Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MMTKVLGMAP VLGPRPPQEQ VGPLMVKVEE KEEKGKYLPS LEMFRQRFRQ FGYHDTPGPR EALSQLRVLC CEWLRPEIHT KEQILELLVL EQFLTILPQE LQAWVQEHCP ESAEEAVTLL EDLERELDEP GHQVSTPPNE QKPVWEKISS SGTAKESPSS MQPQPLETSH KYESWGPLYI QESGEEQEFA QDPRKVRDCR LSTQHEESAD EQKGSEAEGL KGDIISVIIA NKPEASLERQ CVNLENEKGT KPPLQEAGSK KGRESVPTKP TPGERRYICA ECGKAFSNSS NLTKHRRTHT GEKPYVCTKC GKAFSHSSNL TLHYRTHLVD RPYDCKCGKA FGQSSDLLKH QRMHTEEAPY QCKDCGKAFS GKGSLIRHYR IHTGEKPYQC NECGKSFSQH AGLSSHQRLH TGEKPYKCKE CGKAFNHSSN FNKHHRIHTG EKPYWCHHCG KTFCSKSNLS KHQRVHTGEG EAP
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Renal carcinoma antigen NY-REN-21; Zfp-38; Zinc finger and SCAN domain-containing protein 21; zinc finger protein 38 (KOX 25); Zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen
Gene Aliases: NY-REN-21; ZFP38; Zipro1; ZNF38; ZSCAN21
UniProt ID: (Human) Q9Y5A6
Entrez Gene ID: (Human) 7589
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support