Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAKLGKSSS LLTQTSEECN GILTEKMEEE EQTCDPDSSL HWSSSYSPET FRQQFRQFGY QDSPGPHEAL SRLWELCHLW LRPEVHTKEQ ILELLVLEQF LAILPKELQA WVQKHHPENG EETVTMLEDV ERELDGPKQI FFGRRKDMIA EKLAPSEITE ELPSSQLMPV KKQLQGASWE LQSLRPHDED IKTTNVKSAS RQKTSLGIEL HCNVSNILHM NGSQSSTYRG TYEQDGRFEK RQGNPSWKKQ QKCDECGKIF SQSSALILHQ RIHSGKKPYA CDECAKAFSR SAILIQHRRT HTDSKYEHAI AEAQKNMYFK TRLKCPRSLS GGR
Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and SCAN domain-containing protein 14; Zinc finger protein 396
Gene Aliases: ZNF396; ZSCAN14
UniProt ID: (Human) Q96N95
Entrez Gene ID: (Human) 252884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support