Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Recommended positive controls: 293T, A431, Jurakt, Raji.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and SCAN domain-containing protein 14; Zinc finger protein 396
Gene Aliases: ZNF396; ZSCAN14
UniProt ID: (Human) Q96N95
Entrez Gene ID: (Human) 252884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support