Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: GNNFPHSPSS TKNENAKTGA NSCEHDHYEK HLSHKQAPTH HQKIHPEEKL YVCTECVMGF TQKSHLFEHQ RIHAGEKSRE CDKSNKVFPQ KPQVDVHPSV *
This gene product is a likely zinc finger family transcription factor. It contains KRAB-A and KRAB-B domains that act as transcriptional repressors in related proteins, and multiple zinc finger DNA binding motifs and finger linking regions characteristic of the Kruppel family. This gene is part of a gene cluster on chromosome Xp11.23. Several alternatively spliced transcript variants have been described, however, the full-length nature of only some of them is known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mental retardation, X-linked 89; Zinc finger protein 41
Gene Aliases: MRX89; ZNF41
UniProt ID: (Human) P51814
Entrez Gene ID: (Human) 7592
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support