Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: EQFLGTLPP EIQAWVRGQR PGSPEEAAAL VEGLQHDPGQ LLGWITAHVL KQEVLPAAQK TEEPLGSPHP SGTVESPGEG PQDTRIEGSV QLSCSVKEEP NVDGQEVAPS SPPLAAQSPE GHHGHQEPAS TSFHPPRIQE EWGLLDRSQK ELYWDAMLEK YGTVVSLGLP PHQPEAQAQS ELGMLLTGTG VCRSLRSGNE SEGPPGCPEA QPPQGPGPAA WEGLSGAATP APTVRPGTPP VPTQPTPAET RLEPAATPRK PYTCEQCGRG FDWKSVFVIH HRTHTSGPGV QSPGLATGES TEKPPQGEVA FPHHPRHSLT GPRSYPCEEC GCSFSWKSQL VIHRKSHRPE VP (72-422 aa encoded by BC017206)
ZNF446 contains a KRAB and three C(2)H(2) zinc fingers. ZNF446 is a transcription repressor when fused to GAL4 DNA-binding domain and co-transfected with VP-16. Overexpression of ZNF446 in COS-7 cells inhibits the transcriptional activities of SRE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20626; Zinc finger protein 446; Zinc finger protein with KRAB and SCAN domains 20
Gene Aliases: ZKSCAN20; ZNF446; ZSCAN30; ZSCAN52
UniProt ID: (Human) Q9NWS9
Entrez Gene ID: (Human) 55663
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support