Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP762N2316; gene trap insertion site 4-2; ZFPIP; Zinc finger PBX1-interacting protein; Zinc finger protein 462
Gene Aliases: 6030417H05; 9430078C22Rik; Gt4-2; KIAA1803; Zfp462; Zfpip; ZNF462
UniProt ID: (Human) Q96JM2, (Mouse) B1AWL2
Entrez Gene ID: (Human) 58499, (Mouse) 242466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support