Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control:HeLa cells.
Cellular Location: Nucleus.
Transcriptional repressor. Plays a role in oligodendrocyte differentiation, together with OLIG2. Mediates Notch signaling-activated formation of oligodendrocyte precursors. Promotes differentiation of adult neural stem progenitor cells (NSPCs) into mature oligodendrocytes and contributes to remyelination following nerve injury. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 488
Gene Aliases: ZNF488
UniProt ID: (Human) Q96MN9
Entrez Gene ID: (Human) 118738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support