Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: SCVLCCGDLE ATALGRCDHP VCYRCSTKMR VLCEQRYCAV CREELRQVVF GKKLPAFATI PIHQLQHEKK YDIYFADGKV YALYRQLLQH ECPRCPELP
ZNF598 (E3 ubiquitin-protein ligase ZNF598) plays a key role in the ribosome quality control (RQC). RQC is a pathway that takes place when a ribosome has stalled during translation. It is required for ribosomes to terminally stall during translation of poly(A) sequences by mediating monoubiquitination of 40A ribosomal protein RPS10/eS10 and RPS3/uS3. Stalling precludes synthesis of a long poly-lysine tail and initiated the RQC pathway to degrade the potentially detrimental aberrant nascent polypeptide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C6orf149; E3 ubiquitin-protein ligase ZNF598; FLJ00086; ISD11; Zinc finger protein 598
Gene Aliases: ZNF598
UniProt ID: (Human) Q86UK7
Entrez Gene ID: (Human) 90850
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support