Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: vav-1 interacting Kruppel-like protein; Vav-interacting Krueppel-like protein; Zinc finger protein 655
Gene Aliases: VIK; VIK-1; ZNF655
UniProt ID: (Human) Q8N720
Entrez Gene ID: (Human) 79027
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support