Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 704
Gene Aliases: Gig1; ZNF704
UniProt ID: (Human) Q6ZNC4
Entrez Gene ID: (Human) 619279
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support