Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RLCAGENAST PSEPEKFPQV RRQRGAGAGE GEFVCGECGK AFRQSSSLTL
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Human: 100%
May play a role in RNA metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hZNF7; Zinc finger protein 520; Zinc finger protein 74
Gene Aliases: COS52; hZNF7; ZFP520; ZNF520; ZNF74
UniProt ID: (Human) Q16587
Entrez Gene ID: (Human) 7625
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support