Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ZNF746 is a transcription repressor that specifically binds to the 5'-TATTTT[T/G]-3' consensus sequence on promoters and represses transcription of PGC-1-alpha (PPARGC1A), thereby playing a role in regulation of neuron death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PARIS; parkin interacting substrate; Parkin-interacting substrate; parkin-interacting sustrate; Zinc finger protein 746
Gene Aliases: PARIS; ZNF746
UniProt ID: (Human) Q6NUN9
Entrez Gene ID: (Human) 155061
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support