Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRGNYESLVS MDYAISKPDL MSQMERGERP TMQEQEDSEE GETPTDPSAA HDGIVIKIEV QTNDEGSESL ETPEPLMGQV EEHGFQDSEL GDPCGEQPDL DMQEPENTLE ESTEGSSEFS ELKQMLVQQR NCTEGIVIKT EEQDEEEEEE EEDELPQHLQ SLGQLSGRYE ASMYQTPLPG EMSPEGEESP PPLQLGNPTV KRLAPSVHGE RHLSENRGAS SQQQRNRRGE RPFTCMECGK SFRLKINLII HQRNHIKEGP YECAECEISF RHKQQLTLHQ RIHRVRGGCV SPERGPTFNP KHALKPRPKS PSSGSGGGGP KPYKCPECDS SFSHKSSLTK HQITHTGERP YTCPECKKSF RLHISLVIHQ RVHAGKHEVS FICSLCGKSF SRPSHLLRHQ RTHTGERPFK CPECEKSFSE KSKLTNHCRV HSRERPHACP ECGKSFIRKH HLLEHRRIHT GERPYHCAEC GKRFTQKHHL LEHQRAHTGE RPYPCTHCAK CFRYKQSLKY HLRTHTGE
May be involved in transcriptional regulation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 777
Gene Aliases: KIAA1285; ZNF777
UniProt ID: (Human) Q9ULD5
Entrez Gene ID: (Human) 27153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support