Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Immunogen sequence: QGKEHSVSSS REVTSSVLPN DFFSTNPPKA PIIPHSGSIE KAEIHEKVVE RRENTAEALP EGFFDDPEVD ARVRKVDAPK DQM^M
^M
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 16; Coiled-coil domain-containing protein 16; hypothetical protein MGC20398; MGC20398; orphan maintenance of genome 1; Ovus mutant candidate gene 1 protein; Zinc finger protein 830
Gene Aliases: 2410003C20Rik; AV301118; AW048207; CCDC16; OMCG1; Zfp830; ZNF830
UniProt ID: (Human) Q96NB3, (Rat) Q3MHS2, (Mouse) Q8R1N0
Entrez Gene ID: (Human) 91603, (Rat) 497967, (Mouse) 66983
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support