Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: SSSVDFADLP ALFGVPLAPE GIRGYLMEVK PANACHPIEA PRLGNRSLGS IALIRRYDCT FDL
E3 ubiquitin-protein ligase that acts as a negative regulator of NOD2 signaling by mediating ubiquitination and degradation of RIPK2. Also catalyzes ubiquitination and proteasomal degradation of CANX within the endoplasmic reticulum. Could have a role in spermatogenesis (By similarity).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase ZNRF4; KOX18; Nixin; RING finger protein 204; RING-type E3 ubiquitin transferase ZNRF4; Zinc/RING finger protein 4; ZNF139; ZNF36
Gene Aliases: RNF204; SPERIZIN; spzn; Ssrzf1; ZNRF4
UniProt ID: (Human) Q8WWF5
Entrez Gene ID: (Human) 148066
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support