Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ZO-1 Antibodies
            This clone is available in multiple conjugated formats. Search with the clone ID "ZO1-1A12" for more options or explore the "Additional formats" section on this page.

            Invitrogen

            ZO-1 Recombinant Mouse Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ Plus 555

            Advanced Verification
            View all (46) ZO-1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 740002MP555
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            323.00
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite ZO-1 Recombinant Mouse Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ Plus 555

            • Antibody Testing Data (4)
            • Advanced Verification (1)
            Application
            ZO-1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 5

            ZO-1 Antibody (740002MP555) in ICC/IF

            Knockdown of ZO-1 was achieved by transfecting Caco-2 cells with ZO-1 specific siRNA (Silencer® select Product # S14155,S14156). Immunofluorescence analysis was performed on untransfected Caco-2 cells (panel a, d), transfected with non-specific scrambled siRNA (panel b, e) and transfected with ZO-1 specific siRNA (panel c, f). Cells were fixed, permeabilized, and labelled with ZO-1 recombinant mouse monoclonal antibody (ZO1-1A12), Alexa Fluor™ Plus 555 Antibody (740002MP555, 1:1,000 dilution) at 4 degree Celsius overnight. Nuclei (blue) we... View More
            View Product

            ZO-1 Antibody in Immunocytochemistry (ICC/IF)
            ZO-1 Antibody in Immunocytochemistry (ICC/IF)
            ZO-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ZO-1 Antibody in Flow Cytometry (Flow)
            ZO-1 Antibody

            Product Details

            740002MP555

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100
            -

            Immunocytochemistry (ICC/IF)

            1:1,000
            -

            Flow Cytometry (Flow)

            0.063-0.5 µg/test
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG1

            Expression System

            Expi293

            Class

            Recombinant Monoclonal

            Type

            Antibody

            Clone

            ZO1-1A12

            Immunogen

            Human recombinant ZO-1 fusion protein encompassing amino acids 334-634
            View immunogen

            Conjugate

            Alexa Fluor™ Plus 555
            • Unconjugated
            • Alexa Fluor Plus 488 
            • Alexa Fluor Plus 647 
            • Request custom conjugation

            Excitation/Emission Max

            558/572 nm View spectra spectra

            Form

            Liquid

            Concentration

            1.0 mg/mL

            Purification

            Protein A/G

            Storage buffer

            proprietary buffer, pH 6.5

            Contains

            0.008% Bromonitrodioxane, 0.008% Methylisothiazolone

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Wet ice

            RRID

            AB_3093158

            Product Specific Information

            Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro.

            Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.

            Applications Tested: This ZO-1 antibody has been tested by immunohistochemistry of mouse testis tissue and immunocytochemistry and flow cytometric analysis of Caco-2 cells. This may be used at 0.063-0.5 µg per test for flow cytometry. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Excitation: 553 nm; Emission: 568 nm; Laser: Yellow Laser

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Immunocytochemistry
            • Additional Formats
            • Mounting Media ICC
            • Organelle Markers ICC
            • Blockers ICC
            • Counter Stains ICC
            • Labeling Kits ICC
            Catalog #
            740002MP647
            Invitrogen
            ZO-1 Recombinant Mouse Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ Plus 647
            Unit Size :
            50 µL
            Catalog #
            740002M
            Invitrogen
            ZO-1 Recombinant Mouse Monoclonal Antibody (ZO1-1A12)
            Unit Size :
            100 µL
            Catalog #
            740002MP488
            Invitrogen
            ZO-1 Recombinant Mouse Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ Plus 488
            Unit Size :
            50 µL

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: DKFZp686M05161; MGC133289; OTTHUMP00000174520; Tight junction protein 1; Tight junction protein ZO-1; zona occludens 1; Zona occludens protein 1; zonula occludens 1 protein; Zonula occludens protein 1

            View more View less

            Gene Aliases: TJP1; ZO-1; ZO1

            View more View less

            UniProt ID: (Human) Q07157

            View more View less

            Entrez Gene ID: (Human) 7082

            View more View less

            Function(s)

            protein binding calmodulin binding protein domain specific binding cadherin binding involved in cell-cell adhesion tight junction

            Process(es)

            blastocyst formation cell-cell junction assembly sensory perception of sound hippo signaling establishment of endothelial intestinal barrier cell-cell adhesion cell-cell signaling involved in cell-cell junction organization
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-p9qw4:80/100.66.78.181:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0