Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: GRQLQCEFCP VTRWKMAICG LFEIQQCPRG KHCNFLHVFR NPNNEFWEAN RDIYLSPDRT GSSFGKNSER RERMGHHDDY YSRLRGRRNP SPDHSYKRNG
This gene encodes an essential splicing factor. The encoded protein associates with the U2 auxiliary factor heterodimer, which is required for the recognition of a functional 3' splice site in pre-mRNA splicing, and may play a role in network interactions during spliceosome assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2; Renal carcinoma antigen NY-REN-20; U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2; U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2; U2(RNU2) small nuclear RNA auxiliary factor 1-like 2; U2AF35-related protein; URP; zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2; zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2
Gene Aliases: U2AF1-RS2; U2AF1L2; U2AF1RS2; URP; ZC3H22; ZRSR2
UniProt ID: (Human) Q15696
Entrez Gene ID: (Human) 8233
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support