Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: CSRMFKHARS LSSHQRTHLN KKSELLCVTC QKMFKRVSDR RTHEIIHMPE KPFKCSTCEK SFSHKTNLKS HEMIHTGEMP YVCSLCSRRF RQSSTYHRHL RNYHRSD
Embryonic stem (ES) cell-specific transcription factor required to regulate ES cell pluripotency. Binds telomeres and plays a key role in genomic stability in ES cells by regulating telomere elongation. Acts as an activator of spontaneous telomere sister chromatid exchange (T-SCE) and telomere elongation in undifferentiated ES cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and SCAN domain containing protein 4C
Gene Aliases: Gm397; XM_142517; Zscan4c; Zscan4d
UniProt ID: (Mouse) Q80VJ6
Entrez Gene ID: (Mouse) 245109
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support