Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control:HeLa or U87 cells. Ubiquitous tissue expression.
Cellular Location: Nucleus.
ZSCAN5A may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: zinc finger and SCAN domain containing 5; Zinc finger and SCAN domain-containing protein 5A; Zinc finger protein 495
Gene Aliases: ZNF495; ZSCAN5; ZSCAN5A
UniProt ID: (Human) Q9BUG6
Entrez Gene ID: (Human) 79149
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support