Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MLRRGYKASE RRRHLSERLS WHQDQALSSS IYLLREMGPT GFLLREEEPE YMDFRVFLGN PHVCNCSTFP KGGELCKHIC WVLLKKFKLP RNHESALQL
E3 ubiquitin-protein ligase involved in the regulation of Fas-, DR3- and DR4-mediated apoptosis. Functions in conjunction with the UBE2D1, UBE2D3 and UBE2E1 E2 ubiquitin-conjugating enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase ZSWIM2; MEKK1-related protein X; MEX; RING-type E3 ubiquitin transferase ZSWIM2; Zinc finger SWIM domain-containing protein 2; zinc finger, SWIM domain containing 2; zinc finger, SWIM-type containing 2; ZZ-type zinc finger-containing protein 2
Gene Aliases: MEX; ZSWIM2; ZZZ2
UniProt ID: (Human) Q8NEG5
Entrez Gene ID: (Human) 151112
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support