Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDLAALFSD TPANASGSAG GSDEALNSGI LTIDVTSVSS SLGGNLPANN SSLGPMEPLV LVAHSDIPPS LDSPLVLGTA ATVLQQGSFS VDDVQTVSAG ALGCLVALPM KNLSDDPLAL TSNSNLAAHI TTPTSSSTPR ENASVPELLA PIKVEPDSPS RPGAVGQQEG SHGLPQSTLP SPAEQHGAQD TELSAGTGNF YLESGGSART DYRAIQLAKE KKQRGAGSNA GASQSTQRKI KEGKMSPPHF HASQNSWLCG SLVVPSGGRP GPAPAAGVQC GAQGVQVQLV QDDPSGEGVL PSARGPATFL PFLTVDLPVY VLQEVLPSSG GPAGPEATQF PGSTINLQDL Q (509-858 aa encoded by BC002940)
ZXDC belongs to the ZXD family. It contains 10 C2H2-type zinc fingers. ZXDC cooperates with CIITA to promote transcription of MHC class I and MHC class II genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SERH2790; Zinc finger protein ZXDC; ZXD-like zinc finger protein
Gene Aliases: A930012H20; B930086F11Rik; BC003332; ZXDC; ZXDL
UniProt ID: (Human) Q2QGD7, (Mouse) Q8C8V1
Entrez Gene ID: (Human) 79364, (Mouse) 80292
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support