Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAAPRPSPAI SVSVSAPAFY APQEKFGPVV APKPKVNPFR PGDSEPPPAP GAQRAQMGRV GEIPPPPPED FPLPPPPLAG DGDDAEGALG GAFPPPPPPI EESFPPAPLE EEIFPSPPPP PEEEGGPEAP IPPPPQPREK VSSIDLEIDS LSSLLDDMTK NDPFKARVSS GYVPPPVATP FSSKSSTKPA AGGTAPLPPW KSPSSSQPLP QVPAPAQSQT QFHVQPQPQP KPQVQLHVQS QTQPVSLANT QPRGPPASSP APAPKFSPVT PKFTPVASKF SPGAPGGSGS QPNQKLGHPE ALSAGTGSPQ PPSFTYAQQR EKPRVQEKQH PVPPPAQNQN QVRSPGAPGP LTLKEVEELE QLTQQLMQDM EHPQRQNVAV NELCGRCHQP LARAQPAVRA LGQLFHIACF TCHQCAQQLQ GQQFYSLEGA PYCEGCYTDT LEKCNTCGEP ITDRMLRATG KAYHPHCFTC VVCARPLEGT SFIVDQANRP HCVPDYHKQY APRCSVCSEP IMPEPGRDET VRVVALDKNF HMKCYECEDC GKPLSIEADD NGCFPLDGHV LCRKCHTARA QT
Zyxin is involved in focal adhesions, which are actin-rich structures that enable cells to adhere to the extracellular matrix and at which protein complexes involved in signal transduction assemble. Zyxin is a zinc-binding phosphoprotein that concentrates at focal adhesions and along the actin cytoskeleton. Zyxin has an N-terminal proline-rich domain and three LIM domains in its C-terminal half. The proline-rich domain may interact with SH3 domains of proteins involved in signal transduction pathways while the LIM domains are likely involved in protein-protein binding. Zyxin may function as a messenger in the signal transduction pathway that mediates adhesion-stimulated changes in gene expression and may modulate the cytoskeletal organization of actin bundles. Alternative splicing results in multiple transcript variants that encode the same isoform.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zyxin; Zyxin-2
Gene Aliases: ESP-2; HED-2; ZYX
UniProt ID: (Human) Q15942
Entrez Gene ID: (Human) 7791
Molecular Function: actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support