Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YPQRVVQLP EIVWDQYTHS LGNFEREFKN RKRHTRRVKL VFGKVGLPAR PKSPLDPKKD GESLSYSMLP LSDGPEGSSS RPQMIRGRLC DDTKPETFNQ LWTVEEQKKL EQLLIKYPPE EVESRRWQKI ADELGNRTAK QVASQVQKYF IKLTKAGIPV PGRTPNLCIY SKKSSTSRRQ HPLNKHLFKP STFMTSHEPP VYMDEDDDRS CFHSHMNTAV EDASDDESIP IMYRNLPEYK ELLQFKKLKK QKLQQMQAES GFVQHVGFKC DNCGIEPIQG VRWHCQDCPP EMSLDFCDSC SDCLHETDIH KEDHQLEPIY RSETFLDRDY CVSQGTSYNY LDPNYFPANR (555-903 aa encoded by BC035079)
Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATAC component 1 homolog; DKFZP564I052; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3
Gene Aliases: ATAC1; ZZZ3
UniProt ID: (Human) Q8IYH5
Entrez Gene ID: (Human) 26009
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support