Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MVRMVPVLL SLLLLLGPAV PQENQDGRYS LTYIYTGLSK HVEDVPAFQA LGSLNDLQFF RYNSKDRKSQ PMGLWRQVEG MEDWKQDSQL QKAREDIFME TLKDIVEYYN DSNGSHVLQG RFGCEIENNR SSGAFWKYYY DGKDYIEFNK EIPAWVPFDP AAQITKQKWE AEPVYVQRAK AYLEEECPAT LRKYLKYSKN ILDRQDPPSV VVTSHQAPGE KKKLKCLAYD FYPGKIDVHW TRAGEVQEPE LRGDVLHNGN GTYQSWVVVA VPPQDTAPYS CHVQHSSLAQ PLVVPWEAS (1-298 aa encoded by BC033830 )
AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-2-glycoprotein, zinc; testicular tissue protein Li 227; Zinc a 2 glycoprotein; Zinc a2glycoprotein; Zinc alpha2glycoprotein; Zinc α 2 glycoprotein; Zinc α2glycoprotein; Zinc-alpha-2-glycoprotein; Zn a 2 glycoprotein; Zn a 2 GP; Zn alpha 2 glycoprotein; Zn alpha 2 GP; Zn α 2 glycoprotein; Zn α 2 GP; zn-alpha-2-glycoprotein; Zn-alpha-2-GP; Zn-alpha2-glycoprotein
Gene Aliases: AZGP1; ZA2G; ZAG; ZNGP1
UniProt ID: (Human) P25311
Entrez Gene ID: (Human) 563
Molecular Function: major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support