Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Slc30a3 encodes a protein that is involved in accumulation of zinc in synaptic vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Probable proton-coupled zinc antiporter SLC30A3; solute carrier family 30 (zinc transporter), member 3; Solute carrier family 30 member 3; solute carrier family 30 protein; Zinc transporter 3; zinc transporter ZnT-3; ZnT-3
Gene Aliases: SLC30A3; ZNT3
UniProt ID: (Human) Q99726, (Rat) Q6QIX3
Entrez Gene ID: (Pig) 100169931, (Human) 7781, (Rat) 366568
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support