Product References
SIRT2 promotes cell proliferation and migration through mediating ERK1/2 activation and lactosylceramide accumulation in prostate cancer.
The Prostate
Lin R,Yang Y,Wu E,Zhou M,Wang S,Zhang Q
Sun Jan 01 00:00:00 UTC 2023
Oral exposure to polystyrene nanoplastics reduced male fertility and even caused male infertility by inducing testicular and sperm toxicities in mice.
Journal of hazardous materials
Xu W,Yuan Y,Tian Y,Cheng C,Chen Y,Zeng L,Yuan Y,Li D,Zheng L,Luo T
Sat Jul 15 00:00:00 UTC 2023
Scanning electron microscopy of human islet cilia.
bioRxiv : the preprint server for biology
Polino AJ,Sviben S,Melena I,Piston DW,Hughes J
Wed Feb 15 00:00:00 UTC 2023
Optimization of Primary Human Bronchial Epithelial 3D Cell Culture with Donor-Matched Fibroblasts and Comparison of Two Different Culture Media.
International journal of molecular sciences
Maurer J,Walles T,Wiese-Rischke C
Sat Feb 18 00:00:00 UTC 2023
Superenhancer activation of KLHDC8A drives glioma ciliation and hedgehog signaling.
The Journal of clinical investigation
Lee D,Gimple RC,Wu X,Prager BC,Qiu Z,Wu Q,Daggubati V,Mariappan A,Gopalakrishnan J,Sarkisian MR,Raleigh DR,Rich JN
Tue Jan 17 00:00:00 UTC 2023
Superenhancer activation of KLHDC8A drives glioma ciliation and hedgehog signaling.
The Journal of clinical investigation
Lee D,Gimple RC,Wu X,Prager BC,Qiu Z,Wu Q,Daggubati V,Mariappan A,Gopalakrishnan J,Sarkisian MR,Raleigh DR,Rich JN
Tue Jan 17 00:00:00 UTC 2023
Distribution of ciliary adaptor proteins tubby and TULP3 in the organ of Corti.
Frontiers in neuroscience
Lindner LA,Derstroff D,Oliver D,Reimann K
Sun May 07 00:00:00 UTC 2023
Homozygous frameshift variant in POC1B causes male infertility with oligoasthenoteratozoospermia in human and mice.
Human molecular genetics
Hua J,Xu B,Liu W,Shi J,Jiang H,Zha X,Zhang X,Wan Y
Tue Jul 04 00:00:00 UTC 2023
Scanning electron microscopy of human islet cilia.
Proceedings of the National Academy of Sciences of the United States of America
Polino AJ,Sviben S,Melena I,Piston DW,Hughes JW
Tue May 30 00:00:00 UTC 2023
Filamin-A-interacting protein 1 (FILIP1) is a dual compartment protein linking myofibrils and microtubules during myogenic differentiation and upon mechanical stress.
Cell and tissue research
Grande V,Schuld J,van der Ven PFM,Gruss OJ,Fürst DO
Sat Jul 01 00:00:00 UTC 2023
NLRP3 Inflammasome Mediates Silica-induced Lung Epithelial Injury and Aberrant Regeneration in Lung Stem/Progenitor Cell-derived Organotypic Models.
International journal of biological sciences
Zhou H,Zhang Q,Huang W,Zhou S,Wang Y,Zeng X,Wang H,Xie W,Kong H
Tue Apr 18 00:00:00 UTC 2023
Endothelial RGS12 governs angiogenesis in inflammatory arthritis by controlling cilia formation and elongation via MYCBP2 signaling.
Cell insight
Yuan G,Yang ST,Yang S
Sat Oct 01 00:00:00 UTC 2022
KIF17 maintains the epithelial phenotype of breast cancer cells and curbs tumour metastasis.
Cancer letters
Li T,Zhai D,Zhang M,Ye R,Kuang X,Shao N,Bi J,Lin Y
Tue Nov 01 00:00:00 UTC 2022
Islet primary cilia motility controls insulin secretion.
Science advances
Cho JH,Li ZA,Zhu L,Muegge BD,Roseman HF,Lee EY,Utterback T,Woodhams LG,Bayly PV,Hughes JW
Fri Sep 23 00:00:00 UTC 2022
HDAC6 Inhibition Alleviates Ischemia- and Cisplatin-Induced Acute Kidney Injury by Promoting Autophagy.
Cells
Shi L,Song Z,Li C,Deng F,Xia Y,Huang J,Wu X,Zhu J
Wed Dec 07 00:00:00 UTC 2022
Novel Compound Heterozygous Mutation in FSIP2 Causes Multiple Morphological Abnormalities of the Sperm Flagella (MMAF) and Male Infertility.
Reproductive sciences (Thousand Oaks, Calif.)
Hou M,Xi Q,Zhu L,Jia W,Liu Z,Wang C,Zhou X,Zhang D,Xing C,Peng X,Luo Y,Jin L,Li Z,Zhang X
Thu Sep 01 00:00:00 UTC 2022
Inhibition of HDAC6 With CAY10603 Ameliorates Diabetic Kidney Disease by Suppressing NLRP3 Inflammasome.
Frontiers in pharmacology
Hou Q,Kan S,Wang Z,Shi J,Zeng C,Yang D,Jiang S,Liu Z
Tue Aug 02 00:00:00 UTC 2022
Microtubule affinity regulating kinase 4 promoted activation of the NLRP3 inflammasome-mediated pyroptosis in periodontitis.
Journal of oral microbiology
Wang L,Pu W,Wang C,Lei L,Li H
Fri Apr 29 00:00:00 UTC 2022
Microtubule affinity regulating kinase 4 promoted activation of the NLRP3 inflammasome-mediated pyroptosis in periodontitis.
Journal of oral microbiology
Wang L,Pu W,Wang C,Lei L,Li H
Fri Apr 29 00:00:00 UTC 2022
Microtubule affinity regulating kinase 4 promoted activation of the NLRP3 inflammasome-mediated pyroptosis in periodontitis.
Journal of oral microbiology
Wang L,Pu W,Wang C,Lei L,Li H
Fri Apr 29 00:00:00 UTC 2022
Microtubule affinity regulating kinase 4 promoted activation of the NLRP3 inflammasome-mediated pyroptosis in periodontitis.
Journal of oral microbiology
Wang L,Pu W,Wang C,Lei L,Li H
Fri Apr 29 00:00:00 UTC 2022
The acetylome of adult mouse sciatic nerve.
Journal of neurochemistry
Shen M,Chen Z,Ming M,Cheng Z,Sun J,Liang Q,Shang T,Zhang Q,Zhou S,Ji Y,Ding F
Mon Aug 01 00:00:00 UTC 2022
Defective autophagy triggered by arterial cyclic stretch promotes neointimal hyperplasia in vein grafts via the p62/nrf2/slc7a11 signaling pathway.
Journal of molecular and cellular cardiology
Chen Y,Bao M,Liu JT,Bao H,Zhang SM,Lou Y,Qi YX
Thu Dec 01 00:00:00 UTC 2022
A Charcot-Marie-Tooth-Causing Mutation in HSPB1 Decreases Cell Adaptation to Repeated Stress by Disrupting Autophagic Clearance of Misfolded Proteins.
Cells
Zhang X,Qiao Y,Han R,Gao Y,Yang X,Zhang Y,Wan Y,Yu W,Pan X,Xing J
Thu Sep 15 00:00:00 UTC 2022
Biallelic mutations in spermatogenesis and centriole-associated 1 like (SPATC1L) cause acephalic spermatozoa syndrome and male infertility.
Asian journal of andrology
Li YZ,Li N,Liu WS,Sha YW,Wu RF,Tang YL,Zhu XS,Wei XL,Zhang XY,Wang YF,Lu ZX,Zhang FX
Mon Jan 24 00:00:00 UTC 2022
Successful outcomes of intracytoplasmic sperm injection-embryo transfer using ejaculated spermatozoa from two Chinese asthenoteratozoospermic brothers with a compound heterozygous FSIP2 mutation.
Andrologia
Yuan Y,Xu WQ,Chen ZY,Chen Y,Zhang L,Zheng L,Luo T,Chen HY
Fri Apr 01 00:00:00 UTC 2022
Ciliogenesis mechanisms mediated by PAK2-ARL13B signaling in brain endothelial cells is responsible for vascular stability.
Biochemical pharmacology
Thirugnanam K,Prabhudesai S,Van Why E,Pan A,Gupta A,Foreman K,Zennadi R,Rarick KR,Nauli SM,Palecek SP,Ramchandran R
Mon Aug 01 00:00:00 UTC 2022
Fluorescence imaging of beta cell primary cilia.
Frontiers in endocrinology
Li ZA,Cho JH,Woodhams LG,Hughes JW
Tue Oct 11 00:00:00 UTC 2022
Tubastatin A Improves Post-Resuscitation Myocardial Dysfunction by Inhibiting NLRP3-Mediated Pyroptosis Through Enhancing Transcription Factor EB Signaling.
Journal of the American Heart Association
Xu J,Zhao X,Jiang X,He L,Wu X,Wang J,Chen Q,Li Y,Zhang M
Tue Apr 05 00:00:00 UTC 2022
SIRT2-knockdown rescues GARS-induced Charcot-Marie-Tooth neuropathy.
Aging cell
Zhao Y,Xie L,Shen C,Qi Q,Qin Y,Xing J,Zhou D,Qi Y,Yan Z,Lin X,Dai R,Lin J,Yu W
Tue Jun 01 00:00:00 UTC 2021
Mutations in DNAH8 contribute to multiple morphological abnormalities of sperm flagella and male infertility.
Acta biochimica et biophysica Sinica
Weng M,Sha Y,Zeng YU,Huang N,Liu W,Zhang X,Zhou H
Fri Mar 26 00:00:00 UTC 2021
The histone deacetylase inhibitor PCI-24781 impairs calcium influx and inhibits proliferation and metastasis in breast cancer.
Theranostics
Yang T,Wang P,Yin X,Zhang J,Huo M,Gao J,Li G,Teng X,Yu H,Huang W,Wang Y
Mon Aug 16 00:00:00 UTC 2021
Phosphosite T674A mutation in kinesin family member 3A fails to reproduce tissue and ciliary defects characteristic of CILK1 loss of function.
Developmental dynamics : an official publication of the American Association of Anatomists
Gailey CD,Wang EJ,Jin L,Ahmadi S,Brautigan DL,Li X,Xu W,Scott MM,Fu Z
Mon Feb 01 00:00:00 UTC 2021
Nasal ciliated cells are primary targets for SARS-CoV-2 replication in the early stage of COVID-19.
The Journal of clinical investigation
Ahn JH,Kim J,Hong SP,Choi SY,Yang MJ,Ju YS,Kim YT,Kim HM,Rahman MDT,Chung MK,Hong SD,Bae H,Lee CS,Koh GY
Thu Jul 01 00:00:00 UTC 2021
Biallelic mutations in KATNAL2 cause male infertility due to oligo-astheno-teratozoospermia.
Clinical genetics
Wei X,Liu W,Zhu X,Li Y,Zhang X,Chen J,Isachenko V,Sha Y,Lu Z
Fri Oct 01 00:00:00 UTC 2021
Biallelic mutations in KATNAL2 cause male infertility due to oligo-astheno-teratozoospermia.
Clinical genetics
Wei X,Liu W,Zhu X,Li Y,Zhang X,Chen J,Isachenko V,Sha Y,Lu Z
Fri Oct 01 00:00:00 UTC 2021
Biallelic mutations of CFAP58 are associated with multiple morphological abnormalities of the sperm flagella.
Clinical genetics
Sha Y,Sha Y,Liu W,Zhu X,Weng M,Zhang X,Wang Y,Zhou H
Mon Mar 01 00:00:00 UTC 2021
Pathogenic Variants in ACTRT1 Cause Acephalic Spermatozoa Syndrome.
Frontiers in cell and developmental biology
Sha Y,Liu W,Li L,Serafimovski M,Isachenko V,Li Y,Chen J,Zhao B,Wang Y,Wei X
Tue Aug 24 00:00:00 UTC 2021
UTX/KDM6A deletion promotes the recovery of spinal cord injury by epigenetically triggering intrinsic neural regeneration.
Molecular therapy. Methods & clinical development
Guo Z,Li C,Cao Y,Qin T,Jiang L,Xu Y,Li M,Luo Z,Hu J,Lu H
Fri Mar 12 00:00:00 UTC 2021
UTX/KDM6A deletion promotes the recovery of spinal cord injury by epigenetically triggering intrinsic neural regeneration.
Molecular therapy. Methods & clinical development
Guo Z,Li C,Cao Y,Qin T,Jiang L,Xu Y,Li M,Luo Z,Hu J,Lu H
Fri Mar 12 00:00:00 UTC 2021
Analysis of Key Distinct Biological Characteristics of Human Placenta-Derived Mesenchymal Stromal Cells and Individual Heterogeneity Attributing to Donors.
Cells, tissues, organs
Tai C,Wang L,Xie Y,Gao T,Huang F,Wang B
Mon Jan 17 00:00:00 UTC 2022
Sub-toxic levels of cobalt ions impair chondrocyte mechanostranduction via HDAC6-dependent primary cilia shortening.
Biochemical and biophysical research communications
Wu H,Wang Z,Liu S,Meng H,Liu S,Fu S
Fri Mar 12 00:00:00 UTC 2021
HDAC11 promotes meiotic apparatus assembly during mouse oocyte maturation via decreasing H4K16 and α-tubulin acetylation.
Cell cycle (Georgetown, Tex.)
Sui L,Zhang S,Huang R,Li Z
Sat Feb 01 00:00:00 UTC 2020
Photo-triggered self-destructive ROS-responsive nanoparticles of high paclitaxel/chlorin e6 co-loading capacity for synergetic chemo-photodynamic therapy.
Journal of controlled release : official journal of the Controlled Release Society
Yang X,Shi X,Zhang Y,Xu J,Ji J,Ye L,Yi F,Zhai G
Fri Jul 10 00:00:00 UTC 2020
HDAC6 Regulates the Fusion of Autophagosome and Lysosome to Involve in Odontoblast Differentiation.
Frontiers in cell and developmental biology
Zhan Y,Wang H,Zhang L,Pei F,Chen Z
Fri Dec 18 00:00:00 UTC 2020
HDAC6 Regulates the Fusion of Autophagosome and Lysosome to Involve in Odontoblast Differentiation.
Frontiers in cell and developmental biology
Zhan Y,Wang H,Zhang L,Pei F,Chen Z
Fri Dec 18 00:00:00 UTC 2020
Sirtuin 2 Inhibition Attenuates Sevoflurane-Induced Learning and Memory Deficits in Developing Rats via Modulating Microglial Activation.
Cellular and molecular neurobiology
Wu Z,Zhang Y,Zhang Y,Zhao P
Wed Apr 01 00:00:00 UTC 2020
Improved early development potence of in vitro fertilization embryos by treatment with tubacin increasing acetylated tubulin of matured porcine oocytes.
Mechanisms of development
Sun J,Cui K,Li ZP,Gao B,Huang B,Liu Q,Shi D
Tue Dec 01 00:00:00 UTC 2020
Mutations of MAP1B encoding a microtubule-associated phosphoprotein cause sensorineural hearing loss.
JCI insight
Cui L,Zheng J,Zhao Q,Chen JR,Liu H,Peng G,Wu Y,Chen C,He Q,Shi H,Yin S,Friedman RA,Chen Y,Guan MX
Thu Dec 03 00:00:00 UTC 2020
Alternatively Polarized Macrophages Regulate the Growth and Differentiation of Ependymal Stem Cells through the SIRT2 Pathway.
Experimental neurobiology
Ma Y,Deng M,Zhao XQ,Liu M
Thu Apr 30 00:00:00 UTC 2020
UNG2 deacetylation confers cancer cell resistance to hydrogen peroxide-induced cytotoxicity.
Free radical biology & medicine
Bao Y,Tong L,Song B,Liu G,Zhu Q,Lu X,Zhang J,Lu YF,Wen H,Tian Y,Sun Y,Zhu WG
Fri Nov 20 00:00:00 UTC 2020
MiR-107 confers chemoresistance to colorectal cancer by targeting calcium-binding protein 39.
British journal of cancer
Liang Y,Zhu D,Hou L,Wang Y,Huang X,Zhou C,Zhu L,Wang Y,Li L,Gu Y,Luo M,Wang J,Meng X
Sun Mar 01 00:00:00 UTC 2020
Loss-of-function mutations in centrosomal protein 112 is associated with human acephalic spermatozoa phenotype.
Clinical genetics
Sha Y,Wang X,Yuan J,Zhu X,Su Z,Zhang X,Xu X,Wei X
Sat Feb 01 00:00:00 UTC 2020
EBV-miR-BART12 accelerates migration and invasion in EBV-associated cancer cells by targeting tubulin polymerization-promoting protein 1.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wu Y,Wang D,Wei F,Xiong F,Zhang S,Gong Z,Shi L,Li X,Xiang B,Ma J,Deng H,He Y,Liao Q,Zhang W,Li X,Li Y,Guo C,Zeng Z,Li G,Xiong W
Tue Dec 01 00:00:00 UTC 2020
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 UTC 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 UTC 2019
Biallelic mutations in PMFBP1 cause acephalic spermatozoa.
Clinical genetics
Sha YW,Wang X,Xu X,Ding L,Liu WS,Li P,Su ZY,Chen J,Mei LB,Zheng LK,Wang HL,Kong SB,You M,Wu JF
Fri Feb 01 00:00:00 UTC 2019
Biallelic mutations in PMFBP1 cause acephalic spermatozoa.
Clinical genetics
Sha YW,Wang X,Xu X,Ding L,Liu WS,Li P,Su ZY,Chen J,Mei LB,Zheng LK,Wang HL,Kong SB,You M,Wu JF
Fri Feb 01 00:00:00 UTC 2019
Biallelic mutations in PMFBP1 cause acephalic spermatozoa.
Clinical genetics
Sha YW,Wang X,Xu X,Ding L,Liu WS,Li P,Su ZY,Chen J,Mei LB,Zheng LK,Wang HL,Kong SB,You M,Wu JF
Fri Feb 01 00:00:00 UTC 2019
SIRT2 Inhibition Results in Meiotic Arrest, Mitochondrial Dysfunction, and Disturbance of Redox Homeostasis during Bovine Oocyte Maturation.
International journal of molecular sciences
Xu D,Wu L,Jiang X,Yang L,Cheng J,Chen H,Hua R,Geng G,Yang L,Li Q
Mon Mar 18 00:00:00 UTC 2019
The selective HDAC6 inhibitor Nexturastat A induces apoptosis, overcomes drug resistance and inhibits tumor growth in multiple myeloma.
Bioscience reports
Sun X,Xie Y,Sun X,Yao Y,Li H,Li Z,Yao R,Xu K
Fri Mar 29 00:00:00 UTC 2019
Stabilized Peptide HDAC Inhibitors Derived from HDAC1 Substrate H3K56 for the Treatment of Cancer Stem-Like Cells In Vivo.
Cancer research
Wang D,Li W,Zhao R,Chen L,Liu N,Tian Y,Zhao H,Xie M,Lu F,Fang Q,Liang W,Yin F,Li Z
Mon Apr 15 00:00:00 UTC 2019
Ttc39c is upregulated during skeletal muscle atrophy and modulates ERK1/2 MAP kinase and hedgehog signaling.
Journal of cellular physiology
Hayes CS,Labuzan SA,Menke JA,Haddock AN,Waddell DS
Sun Dec 01 00:00:00 UTC 2019
EIF4G1 is a novel candidate gene associated with severe asthenozoospermia.
Molecular genetics & genomic medicine
Sha Y,Liu W,Huang X,Li Y,Ji Z,Mei L,Lin S,Kong S,Lu J,Kong L,Zhu X,Lu Z,Ding L
Thu Aug 01 00:00:00 UTC 2019
EIF4G1 is a novel candidate gene associated with severe asthenozoospermia.
Molecular genetics & genomic medicine
Sha Y,Liu W,Huang X,Li Y,Ji Z,Mei L,Lin S,Kong S,Lu J,Kong L,Zhu X,Lu Z,Ding L
Thu Aug 01 00:00:00 UTC 2019
Biallelic mutations in Sperm flagellum 2 cause human multiple morphological abnormalities of the sperm flagella (MMAF) phenotype.
Clinical genetics
Sha Y,Liu W,Wei X,Zhu X,Luo X,Liang L,Guo T
Fri Nov 01 00:00:00 UTC 2019
Biallelic mutations in Sperm flagellum 2 cause human multiple morphological abnormalities of the sperm flagella (MMAF) phenotype.
Clinical genetics
Sha Y,Liu W,Wei X,Zhu X,Luo X,Liang L,Guo T
Fri Nov 01 00:00:00 UTC 2019
Novel Mutations in CFAP44 and CFAP43 Cause Multiple Morphological Abnormalities of the Sperm Flagella (MMAF).
Reproductive sciences (Thousand Oaks, Calif.)
Sha YW,Wang X,Xu X,Su ZY,Cui Y,Mei LB,Huang XJ,Chen J,He XM,Ji ZY,Bao H,Yang X,Li P,Li L
Tue Jan 01 00:00:00 UTC 2019
A virus-induced kidney disease model based on organ-on-a-chip: Pathogenesis exploration of virus-related renal dysfunctions.
Biomaterials
Wang J,Wang C,Xu N,Liu ZF,Pang DW,Zhang ZL
Tue Oct 01 00:00:00 UTC 2019
Interaction of hnRNP K with MAP 1B-LC1 promotes TGF-β1-mediated epithelial to mesenchymal transition in lung cancer cells.
BMC cancer
Li L,Yan S,Zhang H,Zhang M,Huang G,Chen M
Fri Sep 06 00:00:00 UTC 2019
CCDC9 is identified as a novel candidate gene of severe asthenozoospermia.
Systems biology in reproductive medicine
Sha Y,Xu Y,Wei X,Liu W,Mei L,Lin S,Ji Z,Wang X,Su Z,Qiu P,Chen J,Wang X
Sun Dec 01 00:00:00 UTC 2019
CCDC9 is identified as a novel candidate gene of severe asthenozoospermia.
Systems biology in reproductive medicine
Sha Y,Xu Y,Wei X,Liu W,Mei L,Lin S,Ji Z,Wang X,Su Z,Qiu P,Chen J,Wang X
Sun Dec 01 00:00:00 UTC 2019
Novel IFT140 variants cause spermatogenic dysfunction in humans.
Molecular genetics & genomic medicine
Wang X,Sha YW,Wang WT,Cui YQ,Chen J,Yan W,Hou XT,Mei LB,Yu CC,Wang J
Sun Sep 01 00:00:00 UTC 2019
Myoepithelial Cells of Submucosal Glands Can Function as Reserve Stem Cells to Regenerate Airways after Injury.
Cell stem cell
Tata A,Kobayashi Y,Chow RD,Tran J,Desai A,Massri AJ,McCord TJ,Gunn MD,Tata PR
Thu May 03 00:00:00 UTC 2018
Myoepithelial Cells of Submucosal Glands Can Function as Reserve Stem Cells to Regenerate Airways after Injury.
Cell stem cell
Tata A,Kobayashi Y,Chow RD,Tran J,Desai A,Massri AJ,McCord TJ,Gunn MD,Tata PR
Thu May 03 00:00:00 UTC 2018
C11orf70 Mutations Disrupting the Intraflagellar Transport-Dependent Assembly of Multiple Axonemal Dyneins Cause Primary Ciliary Dyskinesia.
American journal of human genetics
Fassad MR,Shoemark A,le Borgne P,Koll F,Patel M,Dixon M,Hayward J,Richardson C,Frost E,Jenkins L,Cullup T,Chung EMK,Lemullois M,Aubusson-Fleury A,Hogg C,Mitchell DR,Tassin AM,Mitchison HM
Thu May 03 00:00:00 UTC 2018
Gq activity- and β-arrestin-1 scaffolding-mediated ADGRG2/CFTR coupling are required for male fertility.
eLife
Zhang DL,Sun YJ,Ma ML,Wang YJ,Lin H,Li RR,Liang ZL,Gao Y,Yang Z,He DF,Lin A,Mo H,Lu YJ,Li MJ,Kong W,Chung KY,Yi F,Li JY,Qin YY,Li J,Thomsen ARB,Kahsai AW,Chen ZJ,Xu ZG,Liu M,Li D,Yu X,Sun JP
Fri Feb 02 00:00:00 UTC 2018
Gq activity- and β-arrestin-1 scaffolding-mediated ADGRG2/CFTR coupling are required for male fertility.
eLife
Zhang DL,Sun YJ,Ma ML,Wang YJ,Lin H,Li RR,Liang ZL,Gao Y,Yang Z,He DF,Lin A,Mo H,Lu YJ,Li MJ,Kong W,Chung KY,Yi F,Li JY,Qin YY,Li J,Thomsen ARB,Kahsai AW,Chen ZJ,Xu ZG,Liu M,Li D,Yu X,Sun JP
Fri Feb 02 00:00:00 UTC 2018
Spermine increases acetylation of tubulins and facilitates autophagic degradation of prion aggregates.
Scientific reports
Phadwal K,Kurian D,Salamat MKF,MacRae VE,Diack AB,Manson JC
Tue Jul 03 00:00:00 UTC 2018
Spermine increases acetylation of tubulins and facilitates autophagic degradation of prion aggregates.
Scientific reports
Phadwal K,Kurian D,Salamat MKF,MacRae VE,Diack AB,Manson JC
Tue Jul 03 00:00:00 UTC 2018
Spermine increases acetylation of tubulins and facilitates autophagic degradation of prion aggregates.
Scientific reports
Phadwal K,Kurian D,Salamat MKF,MacRae VE,Diack AB,Manson JC
Tue Jul 03 00:00:00 UTC 2018
Spermine increases acetylation of tubulins and facilitates autophagic degradation of prion aggregates.
Scientific reports
Phadwal K,Kurian D,Salamat MKF,MacRae VE,Diack AB,Manson JC
Tue Jul 03 00:00:00 UTC 2018
HDAC6 Suppresses Age-Dependent Ectopic Fat Accumulation by Maintaining the Proteostasis of PLIN2 in Drosophila.
Developmental cell
Yan Y,Wang H,Hu M,Jiang L,Wang Y,Liu P,Liang X,Liu J,Li C,Lindström-Battle A,Lam SM,Shui G,Deng WM,Jiao R
Mon Oct 09 00:00:00 UTC 2017
HDAC6 Suppresses Age-Dependent Ectopic Fat Accumulation by Maintaining the Proteostasis of PLIN2 in Drosophila.
Developmental cell
Yan Y,Wang H,Hu M,Jiang L,Wang Y,Liu P,Liang X,Liu J,Li C,Lindström-Battle A,Lam SM,Shui G,Deng WM,Jiao R
Mon Oct 09 00:00:00 UTC 2017
RPGR, a prenylated retinal ciliopathy protein, is targeted to cilia in a prenylation- and PDE6D-dependent manner.
Biology open
Dutta N,Seo S
Thu Sep 15 00:00:00 UTC 2016
RPGR, a prenylated retinal ciliopathy protein, is targeted to cilia in a prenylation- and PDE6D-dependent manner.
Biology open
Dutta N,Seo S
Thu Sep 15 00:00:00 UTC 2016
ProBDNF inhibits collective migration and chemotaxis of rat Schwann cells.
Tissue & cell
Ding YQ,Li XY,Xia GN,Ren HY,Zhou XF,Su BY,Qi JG
Sat Oct 01 00:00:00 UTC 2016
NDUFV2 regulates neuronal migration in the developing cerebral cortex through modulation of the multipolar-bipolar transition.
Brain research
Chen T,Wu Q,Zhang Y,Zhang D
Mon Nov 02 00:00:00 UTC 2015
NDUFV2 regulates neuronal migration in the developing cerebral cortex through modulation of the multipolar-bipolar transition.
Brain research
Chen T,Wu Q,Zhang Y,Zhang D
Mon Nov 02 00:00:00 UTC 2015
NDUFV2 regulates neuronal migration in the developing cerebral cortex through modulation of the multipolar-bipolar transition.
Brain research
Chen T,Wu Q,Zhang Y,Zhang D
Mon Nov 02 00:00:00 UTC 2015
NDUFV2 regulates neuronal migration in the developing cerebral cortex through modulation of the multipolar-bipolar transition.
Brain research
Chen T,Wu Q,Zhang Y,Zhang D
Mon Nov 02 00:00:00 UTC 2015
The oncoprotein HBXIP promotes migration of breast cancer cells via GCN5-mediated microtubule acetylation.
Biochemical and biophysical research communications
Li L,Liu B,Zhang X,Ye L
Fri Mar 13 00:00:00 UTC 2015
The oncoprotein HBXIP promotes migration of breast cancer cells via GCN5-mediated microtubule acetylation.
Biochemical and biophysical research communications
Li L,Liu B,Zhang X,Ye L
Fri Mar 13 00:00:00 UTC 2015
The oncoprotein HBXIP promotes migration of breast cancer cells via GCN5-mediated microtubule acetylation.
Biochemical and biophysical research communications
Li L,Liu B,Zhang X,Ye L
Fri Mar 13 00:00:00 UTC 2015
The oncoprotein HBXIP promotes migration of breast cancer cells via GCN5-mediated microtubule acetylation.
Biochemical and biophysical research communications
Li L,Liu B,Zhang X,Ye L
Fri Mar 13 00:00:00 UTC 2015