Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MUbio
HHF35 reacts with both alpha-muscle and gamma-smooth muscle actin, and therefore reacts with skeletal muscle, cardiac muscle, vascular and visceral smooth muscle cells, pericytes and myoepithelial cells. It is also reactive in myofibroblasts. It does not react with epithelial, endothelial, neural or normal connective tissue cells when applied under the proper conditions to these tissue sections.
The epitope recognized by α-SM1 is highly conserved. The antibody therefore cross-reacts with many species including protochordates, lower craniates and mammals.
The antibody may be stored at 4°C. For prolonged storage prepare appropriate aliquots and store at or below -20°C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.
Actins are highly conserved proteins that are involved in various types of cell motility, and maintenance of the cytoskeleton. In vertebrates, three main groups of actin isoforms, alpha, beta and gamma have been identified. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton, and as mediators of internal cell motility. Actin gamma 1 is a cytoplasmic actin found in nonmuscle cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin gamma; Actin, cytoplasmic 2; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1 propeptide; actin, gamma, cytoplasmic 1; Atcb; cytoplasmic 2; cytoplasmic ACTG1; cytoplasmic type 5; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; epididymis luminal protein 176; gamma 1 propeptide; Gamma-actin; type 5
Gene Aliases: ACT; ACTG; ACTG1; actin; BRWS2; DFNA20; DFNA26; HEL-176
UniProt ID: (Chicken) P53478, (Human) P63261, (Rat) P63259
Entrez Gene ID: (Pig) 397653, (Chicken) 415296, (Human) 71, (Rat) 287876
Molecular Function: actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support