Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 5
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADA1D; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; Alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; Alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; Alpha-adrenergic receptor 1a; Alpha1delta-AR; RA42
Gene Aliases: [a]1d; Adra-1; ADRA1; ADRA1A; ADRA1D; ADRA1R; Adrd1; ALPHA1; alpha1D-AR; DAR; dJ779E11.2; Gpcr8; Spr8
UniProt ID: (Human) P25100, (Rat) P23944
Entrez Gene ID: (Human) 146, (Mouse) 11550, (Rat) 29413
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support