Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-32690 detects alpha 1d Adrenoceptor from human samples.
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADA1D; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; Alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; Alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; Alpha-adrenergic receptor 1a; Alpha1delta-AR
Gene Aliases: ADRA1; ADRA1A; ADRA1D; ADRA1R; ALPHA1; DAR; dJ779E11.2
UniProt ID: (Human) P25100
Entrez Gene ID: (Human) 146
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support