Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Positive test controls include: K-562, Mouse brain, Mouse Spinal cord. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein.
Immunogen sequence: RRTRVPPSRR GPDAVAAPPG GTERRPNGLG PERSAGPGGA EAEPLPTQLN GAPGEPAPAG PRDTDALDLE ESSSSDHAER PPGPRRPERG PRGKGKARAS QVKPGDSLPR RGPGATGIGT PAAGPGEERV GAAKASRWRG R
Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; Alpha-2 adrenergic receptor subtype C10; alpha-2-adrenergic receptor, platelet type; Alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; Alpha-2A adrenoreceptor; alpha-2AAR; alpha-2AAR subtype C10; Alpha-2D adrenergic receptor; alpha2A-adrenergic receptor; CA2-47; OTTHUMP00000058849; subtype alpha2-C10
Gene Aliases: Adra-2; Adra-2a; ADRA2; ADRA2A; ADRA2R; ADRAR; alpha(2A)AR; alpha2-C10; alpha2A; alpha2A-AR; ALPHA2AAR; AW122659; CA2-47; RATRG20; RG20; ZNF32
UniProt ID: (Human) P08913, (Mouse) Q01338, (Rat) P22909
Entrez Gene ID: (Human) 150, (Mouse) 11551, (Rat) 25083
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support