Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 14
Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261, C8D30, NIH-3T3, Rat heart.
Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (94%), Pig (99%), Chicken (98%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it i involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actinin, alpha 2; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin-2; F-actin cross-linking protein
Gene Aliases: 1110008F24Rik; ACTN2; CMD1AA; CMH23
UniProt ID: (Human) P35609, (Mouse) Q9JI91
Entrez Gene ID: (Human) 88, (Pig) 100157406, (Mouse) 11472, (Rat) 291245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support