Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Positive Samples: HeLa, HT-29, MCF7, Mouse lung, Rat brain
Immunogen sequence: MLTKFETKSA RVKGLSFHPK RPWILTSLHN GVIQLWDYRM CTLIDKFDEH DGPVRGIDFH KQQPLFVSGG DDYKIKVWNY KLRRCLFTLL GHLDYIRTTF
In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha coat protein; Alpha-coat protein; Alpha-COP; Coatomer subunit alpha; HEP-COP; HEPCOP; proxenin; xenin
Gene Aliases: AILJK; AU040324; COPA; HEP-COP; xenin
UniProt ID: (Human) P53621, (Mouse) Q8CIE6
Entrez Gene ID: (Human) 1314, (Mouse) 12847, (Rat) 304978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support