Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, mouse kidney.
Predicted reactivity: Mouse (84%), Rat (85%), Pig (85%), Rabbit (89%), Bovine (85%), Guinea pig (83%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel’s syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha ENaC-2; Alpha-ENaC; Alpha-NaCH; amiloride-sensitive epithelial sodium channel; amiloride-sensitive epithelial sodium channel alpha subunit; Amiloride-sensitive sodium channel subunit alpha; amiloride-sensitive sodium channel subunit alpha 2; ENaC alpha; Epithelial Na(+) channel subunit alpha; epithelial sodium channel alpha subunit; FLJ21883; nasal epithelial sodium channel alpha subunit; Nonvoltage-gated sodium channel 1 subunit alpha; SCNEA; sodium channel, non voltage gated 1 alpha subunit; sodium channel, non-voltage-gated 1 alpha subunit; sodium channel, nonvoltage-gated 1 alpha; sodium channel, nonvoltage-gated, type I, alpha polypeptide
Gene Aliases: BESC2; ENaC; ENaCa; ENaCalpha; mENaC; SCNEA; SCNN1; SCNN1A
UniProt ID: (Human) P37088, (Mouse) Q61180
Entrez Gene ID: (Human) 6337, (Mouse) 20276
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support