Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as one of the large subunits of this complex and is a member of the adaptin protein family. This gene is a candidate meningioma gene. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 1 subunit beta-1; adapter-related protein complex 2 beta subunit; adapter-related protein complex 2 subunit beta; adaptin, beta 2 (beta); Adaptor protein complex AP-1 subunit beta-1; adaptor protein complex AP-1, beta 1 subunit; Adaptor protein complex AP-2 subunit beta; adaptor related protein complex 2, beta 1 subunit; adaptor-related protein complex 1 beta 1 subunit; Adaptor-related protein complex 1 subunit beta-1; adaptor-related protein complex 1, beta 1 subunit; Adaptor-related protein complex 2 subunit beta; adaptor-related protein complex 2, beta 1 subunit; adaptor-related protein complex AP-1 beta 1 subunit; adaptor-related protein complex AP-1, beta 1 subunit; AP-1 complex subunit beta-1; AP-2 complex subunit beta; AP105B; beta adaptin; Beta-1-adaptin; Beta-2-adaptin; Beta-adaptin; Beta-adaptin 1; beta-prime adaptin; beta-prime-adaptin; beta1-adaptin; beta2-adaptin; Clathrin assembly protein complex 1 beta large chain; Clathrin assembly protein complex 2 beta large chain; clathrin-associated/assembly/adaptor protein, large, beta 1; Golgi adaptor HA1/AP1 adaptin beta subunit; Mutant line 1660; Plasma membrane adaptor HA2/AP2 adaptin beta subunit; plasma membrane adaptor HA2/AP2 adaptor beta subunit; testicular tissue protein Li 22
Gene Aliases: 1300012O03Rik; ADTB1; ADTB2; AI788979; AP105A; AP105B; AP1B1; AP2-BETA; AP2B1; b2b1660Clo; BAM22; CLAPB1; CLAPB2
UniProt ID: (Human) Q10567, (Human) P63010, (Mouse) O35643, (Rat) P62944, (Mouse) Q9DBG3
Entrez Gene ID: (Human) 162, (Human) 163, (Rat) 29663, (Mouse) 11764, (Rat) 140670, (Mouse) 71770
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support