Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Beta III Tubulin Antibodies

            Invitrogen

            beta-3 Tubulin Polyclonal Antibody

            Advanced Verification
            2 Published Figures
            7 References
            View all (50) Beta III Tubulin antibodies

            Promotions

            View available promotion(s)

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-85639
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            510.00
            100 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite beta-3 Tubulin Polyclonal Antibody

            • Antibody Testing Data (15)
            • Published Figures (2)
            • Advanced Verification (4)
            Application
            beta-3 Tubulin Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 21

            beta-3 Tubulin Antibody (PA5-85639) in ICC/IF

            beta-3 Tubulin Polyclonal Antibody detects beta Tubulin 3/ Tuj1 protein at cytoplasm by immunofluorescent analysis. Sample: DIV9 rat E18 primary cortical neurons were fixed in 4% paraformaldehyde at RT for 15 min. Green: beta Tubulin 3/ Tuj1 protein stained by beta-3 Tubulin Polyclonal Antibody (Product # PA5-85639) diluted at 1:500. Blue: Fluoroshield with DAPI .
            View Product

            beta-3 Tubulin Antibody in Immunocytochemistry (ICC/IF)
            beta-3 Tubulin Antibody in Immunocytochemistry (ICC/IF)
            beta-3 Tubulin Antibody in Immunocytochemistry (ICC/IF)
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunohistochemistry (Frozen) (IHC (F))
            beta-3 Tubulin Antibody in Western Blot (WB)
            beta-3 Tubulin Antibody in Western Blot (WB)
            beta-3 Tubulin Antibody in Western Blot (WB)
            beta-3 Tubulin Antibody in Western Blot (WB)
            beta-3 Tubulin Antibody in Immunoprecipitation (IP)
            beta-3 Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            beta-3 Tubulin Antibody in Immunocytochemistry (ICC/IF)
            beta-3 Tubulin Antibody
            beta-3 Tubulin Antibody
            beta-3 Tubulin Antibody
            beta-3 Tubulin Antibody

            Product Details

            PA5-85639

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:50,000
            View 1 publication 1 publication

            Immunohistochemistry (IHC)

            -
            View 2 publications 2 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:1,000
            View 1 publication 1 publication

            Immunohistochemistry (Frozen) (IHC (F))

            1:100-1:1,000
            -

            Immunocytochemistry (ICC/IF)

            1:100-1:1,000
            View 2 publications 2 publications

            Flow Cytometry (Flow)

            Assay-dependent
            -

            Immunoprecipitation (IP)

            1:100-1:500
            -

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human beta Tubulin 3/ TUJ1. The exact sequence is proprietary.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.53 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7, with 20% glycerol

            Contains

            0.025% ProClin 300

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2792779

            Product Specific Information

            Keep as concentrated solution.

            Positive Control: HeLa, U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, rat brain.

            Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

            Target Information

            The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Immunohistochemistry
            • Secondary Antibodies
            • Isotype Controls
            • Mounting Media IHC
            • Counter Stains IHC
            • Blockers IHC
            • Substrates IHC
            • Labeling Kits IHC

            Bioinformatics

            Protein Aliases: beta 3 tubulin; beta tubulin 3; betaIII-tubulin; class III beta-tubulin; MC1R; Neuron specific beta III Tubulin; Neuron-specific class III beta-tubulin; TUBB; tubulin beta; Tubulin beta-3; Tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; Tubulin beta-III; tubulin, beta 3 class III

            View more View less

            Gene Aliases: 3200002H15Rik; beta-4; CDCBM; CDCBM1; CFEOM3; CFEOM3A; FEOM3; M(beta)3; M(beta)6; TUBB3; TUBB4

            View more View less

            UniProt ID: (Human) Q13509, (Mouse) Q9ERD7, (Rat) Q4QRB4

            View more View less

            Entrez Gene ID: (Human) 10381, (Mouse) 22152, (Rat) 246118

            View more View less

            Function(s)

            GTPase activity structural constituent of cytoskeleton protein binding GTP binding peptide binding nucleotide binding tubulin

            Process(es)

            cytoskeleton organization microtubule-based process mitotic nuclear division axon guidance neuron differentiation protein polymerization metabolic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0