Product References
Mitofusin2 Ameliorated Endoplasmic Reticulum Stress and Mitochondrial Reactive Oxygen Species Through Maintaining Mitochondria-Associated Endoplasmic Reticulum Membrane Integrity in Cisplatin-Induced Acute Kidney Injury.
Antioxidants & redox signaling
Liu YT,Zhang H,Duan SB,Wang JW,Chen H,Zhan M,Zhang W,Li AM,Liu Y,Yang Y,Yang S
Mon Jan 01 00:00:00 UTC 2024
WITHDRAWN: Expression of Tmem41b and MMP13 associated with poor outcome in osteosarcomas.
Neoplasma
Li GH,Liu X,Feng LJ,Zhang L
Sun Aug 20 00:00:00 UTC 2023
Kinsenoside alleviates oxidative stress-induced blood-brain barrier dysfunction via promoting Nrf2/HO-1 pathway in ischemic stroke.
European journal of pharmacology
Qiao N,An Z,Fu Z,Chen X,Tong Q,Zhang Y,Ren H
Thu Jun 15 00:00:00 UTC 2023
Persistent Coxsackievirus B3 Infection in Pancreatic Ductal Cells In Vitro Downregulates Cellular Polyamine Metabolism.
mSphere
Mastrodomenico V,LoMascolo NJ,Firpo MR,Villanueva Guzman MDM,Zaporowski A,Mounce BC
Thu Jun 22 00:00:00 UTC 2023
Exercise induced meteorin-like protects chondrocytes against inflammation and pyroptosis in osteoarthritis by inhibiting PI3K/Akt/NF-κB and NLRP3/caspase-1/GSDMD signaling.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu J,Jia S,Yang Y,Piao L,Wang Z,Jin Z,Bai L
Wed Feb 01 00:00:00 UTC 2023
The Role of LincRNA-EPS/Sirt1/Autophagy Pathway in the Neuroprotection Process by Hydrogen against OGD/R-Induced Hippocampal HT22 Cells Injury.
Journal of personalized medicine
Li YH,Zhang S,Tang L,Feng J,Jia J,Chen Y,Liu L,Zhou J
Mon Apr 03 00:00:00 UTC 2023
Karyopherin Subunit Alpha 1 Enhances the Malignant Behaviors of Colon Cancer Cells via Promoting Nuclear Factor-κB p65 Nuclear Translocation.
Digestive diseases and sciences
Zhao L,Wu D,Qu Q,Li Z,Yin H
Sat Jul 01 00:00:00 UTC 2023
Irisin attenuates acute lung injury by suppressing the pyroptosis of alveolar macrophages.
International journal of molecular medicine
Han Z,Ma J,Han Y,Yuan G,Jiao R,Meng A
Sat Apr 01 00:00:00 UTC 2023
PM2.5 mediates mouse testis Sertoli TM4 cell damage by reducing cellular NAD.
Toxicology mechanisms and methods
Xu P,Ren T,Yang Y
Sun Oct 01 00:00:00 UTC 2023
Repetitive transcranial magnetic stimulation alleviates glial activation through suppressing HMGB1/TLR4 pathway in a rat model of Parkinson's disease.
Molecular biology reports
Han C,Zhang X,Dou K,Yao W,Yao M,Wan Q,Xie A
Tue Aug 01 00:00:00 UTC 2023
IL-33/ST2 mediating systemic inflammation and neuroinflammation through NF-kB participated in the neurocognitive impairment in obstructive sleep apnea.
International immunopharmacology
Shen C,Liu X,Liu T,Ou Y,Ouyang R
Wed Feb 01 00:00:00 UTC 2023
Disruption of COMMD1 accelerates diabetic atherosclerosis by promoting glycolysis.
Diabetes & vascular disease research
Zhang L,Li L,Li Y,Jiang H,Sun Z,Zang G,Qian Y,Shao C,Wang Z
Thu Feb 23 00:00:00 UTC 2023
Anti-glycation and anti-inflammatory activities of anthocyanins from purple vegetables.
Food & function
Gao Q,Ma R,Shi L,Wang S,Liang Y,Zhang Z
Tue Feb 21 00:00:00 UTC 2023
Trans-gnetin H isolated from the seeds of Paeonia species induces autophagy via inhibiting mTORC1 signalling through AMPK activation.
Cell proliferation
Xia C,Wang G,Chen L,Geng H,Yao J,Bai Z,Deng L
Wed Mar 01 00:00:00 UTC 2023
Efficacy of Danggui Buxue decoction on diabetic nephropathy-induced renal fibrosis in rats and possible mechanism.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan
Ye Z,Xian W,Ling GU,Zihang LI,Jingtian Z,Wenkai W,Liang Z,Mei X
Thu Jun 01 00:00:00 UTC 2023
Bacillus Calmette-Guérin-induced interleukin-10 inhibits S100A8/A9 production and hinders development of T helper type 1 memory in mice.
European journal of immunology
Wang Y,Sun Y,Zheng Y,Yang Y,He L,Qu P,Zhou F,Xu X,Bai X,Chen X,Yuan Y,Liu M,Pan Q
Sat Apr 01 00:00:00 UTC 2023
Pachymic acid ameliorates bleomycin-induced pulmonary fibrosis through inhibiting endoplasmic reticulum stress in rats.
Environmental toxicology
Li L,Jin RJ,Ji L
Wed May 10 00:00:00 UTC 2023
Evaluation of Brown Micro-Algae Synergies With Low Dose γ-Radiation Against Chronic Hepatitis Induced by D-Galactosamine in Rats.
Dose-response : a publication of International Hormesis Society
Elsaman S,Elsonbaty SM,Moawed FSM,Hegazy MGA
Fri Apr 21 00:00:00 UTC 2023
Effects of a ketogenic diet on reproductive and metabolic phenotypes in mice with polycystic ovary syndrome†.
Biology of reproduction
Liu S,Yao Q,Li X,Wu H,Sun C,Bai W,Kang J
Tue Apr 11 00:00:00 UTC 2023
CD166-specific CAR-T cells potently target colorectal cancer cells.
Translational oncology
He S,Li S,Guo J,Zeng X,Liang D,Zhu Y,Li Y,Yang D,Zhao X
Sun Jan 01 00:00:00 UTC 2023
Cyasterone inhibits IL-1β-mediated apoptosis and inflammation via the NF-κB and MAPK signaling pathways in rat chondrocytes and ameliorates osteoarthritisin vivo.
Chinese journal of natural medicines
Teng L,Shen Y,Qu Y,Yang L,Yang Y,Jian X,Fan S,Zhang L,Fu Q
Wed Feb 01 00:00:00 UTC 2023
Alpha-kinase1 promotes tubular injury and interstitial inflammation in diabetic nephropathy by canonical pyroptosis pathway.
Biological research
Cui X,Li Y,Yuan S,Huang Y,Chen X,Han Y,Liu Z,Li Z,Xiao Y,Wang Y,Sun L,Liu H,Zhu X
Thu Feb 02 00:00:00 UTC 2023
Elevated PGT promotes proliferation and inhibits cell apoptosis in preeclampsia by Erk signaling pathway.
Molecular and cellular probes
Pang H,Lei D,Huang J,Guo Y,Fan C
Wed Feb 01 00:00:00 UTC 2023
PKM2-mediated neuronal hyperglycolysis enhances the risk of Parkinson's disease in diabetic rats.
Journal of pharmaceutical analysis
Zhao Y,Wang Y,Wu Y,Tao C,Xu R,Chen Y,Qian L,Xu T,Lian X
Wed Feb 01 00:00:00 UTC 2023
Robo4 inhibits gamma radiation-induced permeability of a murine microvascular endothelial cell by regulating the junctions.
Cellular & molecular biology letters
Adzraku SY,Wang G,Cao C,Bao Y,Wang Y,Smith AO,Du Y,Wang H,Li Y,Xu K,Qiao J,Ju W,Zeng L
Mon Jan 16 00:00:00 UTC 2023
BAP18 facilitates CTCF-mediated chromatin accessible to regulate enhancer activity in breast cancer.
Cell death and differentiation
Sun G,Wei Y,Zhou B,Wang M,Luan R,Bai Y,Li H,Wang S,Zheng D,Wang C,Wang S,Zeng K,Liu S,Lin L,He M,Zhang Q,Zhao Y
Mon May 01 00:00:00 UTC 2023
Recombinant humanized collagen type III with high antitumor activity inhibits breast cancer cells autophagy, proliferation, and migration through DDR1.
International journal of biological macromolecules
Liu X,Li H,Wang T,Yang T,Yang X,Guo K,Hu L,Ming J
Sat Jul 15 00:00:00 UTC 2023
Visceral adipose tissue-directed human kallistatin gene therapy improves adipose tissue remodeling and metabolic health in obese mice.
Cellular signalling
Wu Y,Jiang Y,Guo JQ,Yang ZW,Carvalho A,Qian LL,Ji JJ,Ji ZJ,Ma GS,Yao YY
Thu Jun 01 00:00:00 UTC 2023
Remote Ischemic Conditioning Relieves Necrotizing Enterocolitis Through Regulation of Redox and Inflammation.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Zhang Y,Liu Q,Gao K,Tian B,Zhu H,Liu J,Hu Y,Ye C,Guo C
Mon May 01 00:00:00 UTC 2023
Isoginkgetin-loaded reactive oxygen species scavenging nanoparticles ameliorate intervertebral disc degeneration via enhancing autophagy in nucleus pulposus cells.
Journal of nanobiotechnology
Yu H,Teng Y,Ge J,Yang M,Xie H,Wu T,Yan Q,Jia M,Zhu Q,Shen Y,Zhang L,Zou J
Tue Mar 21 00:00:00 UTC 2023
Possible mechanisms associated with immune escape and apoptosis on anti-hepatocellular carcinoma effect of Mu Ji Fang granules.
World journal of gastrointestinal oncology
Zhang YB,Bao YR,Wang S,Li TJ,Tai H,Leng JP,Yang XX,Wang BC,Meng XS
Wed Mar 15 00:00:00 UTC 2023
DDX3X Is Hijacked by Snakehead Vesiculovirus Phosphoprotein To Facilitate Virus Replication via Stabilization of the Phosphoprotein.
Journal of virology
Bei C,Zhang C,Wu H,Feng H,Zhang YA,Tu J
Tue Feb 28 00:00:00 UTC 2023
Tiliroside targets TBK1 to induce ferroptosis and sensitize hepatocellular carcinoma to sorafenib.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yang C,Lu T,Liu M,Yuan X,Li D,Zhang J,Zhou L,Xu M
Wed Mar 01 00:00:00 UTC 2023
Long-term Pu-erh tea consumption improves blue light-induced depression-like behaviors.
Food & function
Zhao S,Hu S,Sun K,Luo L,Zeng L
Mon Mar 06 00:00:00 UTC 2023
Curcumin Alleviates Hepatic Ischemia-Reperfusion Injury by Inhibiting Neutrophil Extracellular Traps Formation.
Journal of investigative surgery : the official journal of the Academy of Surgical Research
Zhu C,Shi S,Jiang P,Huang X,Zhao J,Jin Y,Shen Y,Zhou X,Liu H,Cai J
Fri Dec 01 00:00:00 UTC 2023
The Activation of Reticulophagy by ER Stress through the ATF4-MAP1LC3A-CCPG1 Pathway in Ovarian Granulosa Cells Is Linked to Apoptosis and Necroptosis.
International journal of molecular sciences
Li H,Jing Y,Qu X,Yang J,Pan P,Liu X,Gao H,Pei X,Zhang C,Yang Y
Wed Feb 01 00:00:00 UTC 2023
S-Doped carbonized polymer dots inhibit early myocardial fibrosis by regulating mitochondrial function.
Biomaterials science
Wang Y,Yang M,Zhang J,Ren J,Liu N,Liu B,Lu L,Yang B
Tue Jan 31 00:00:00 UTC 2023
SARS-COV-2 spike protein promotes RPE cell senescence via the ROS/P53/P21 pathway.
Biogerontology
Zhang Y,Peng X,Xue M,Liu J,Shang G,Jiang M,Chen D,Liu B,Wang Y,Jia X,Xu J,Zhang F,Hu Y
Sun Oct 01 00:00:00 UTC 2023
LINC00346 regulates NLRP1-mediated pyroptosis and autophagy via binding to microRNA-637 in vascular endothelium injury.
Cellular signalling
Ge JY,Yan XJ,Yang J,Jin H,Sun ZK,Guo JL,Zhu Y,Wang FF
Fri Sep 01 00:00:00 UTC 2023
Organic extracts in PM2.5 are the major triggers to induce ferroptosis in SH-SY5Y cells.
Ecotoxicology and environmental safety
Guo C,Lyu Y,Xia S,Ren X,Li Z,Tian F,Zheng J
Sun Jan 01 00:00:00 UTC 2023
Dietary alpha-ketoglutarate enhances intestinal immunity by Th17/Treg immune response in piglets after lipopolysaccharide challenge.
Journal of animal science
Liu GM,Lu JJ,Sun WX,Jia G,Zhao H,Chen XL,Tian G,Cai JY,Zhang RN,Wang J
Tue Jan 03 00:00:00 UTC 2023
Oncolytic Hairpin DNA Pair: Selective Cytotoxic Inducer through MicroRNA-Triggered DNA Self-Assembly.
Journal of the American Chemical Society
Morihiro K,Osumi H,Morita S,Hattori T,Baba M,Harada N,Ohashi R,Okamoto A
Wed Jan 11 00:00:00 UTC 2023
Yishen-Qingli-Huoxue formula attenuates renal fibrosis by inhibiting indoxyl sulfate via AhR/snai1 signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Chen C,Sun S,Zhao J,Wu Q,He W,Sun W
Sun Jan 01 00:00:00 UTC 2023
Transcriptomic and functional analyses reveal the molecular mechanisms underlying Fe-mediated tobacco resistance to potato virus Y infection.
Frontiers in plant science
Xu C,Guo H,Li R,Lan X,Zhang Y,Xie Q,Zhu D,Mu Q,Wang Z,An M,Xia Z,Wu Y
Tue Apr 18 00:00:00 UTC 2023
CEMIP promotes small cell lung cancer proliferation by activation of glutamine metabolism via FBXW7/c-Myc-dependent axis.
Biochemical pharmacology
Mo X,Shen X,Mo X,Yu F,Tan W,Deng Z,He J,Luo Z,Chen Z,Yang J
Wed Mar 01 00:00:00 UTC 2023
Neuroprotective effect of a Keap1-Nrf2 Protein-Protein Inter-action inhibitor on cerebral Ischemia/Reperfusion injury.
Bioorganic chemistry
Qi Z,Tong Y,Luo H,Chen M,Zhou N,Chen L
Wed Mar 01 00:00:00 UTC 2023
Hsa_circ_0003596 enhances the development of cell renal clear cell carcinoma through the miR-502-5p/IGF1/PI3K/AKT axis.
The journal of gene medicine
Fang K,Liang S,Liu D,Yi Q,Li Y,Zhu R
Fri Dec 01 00:00:00 UTC 2023
Patrinia villosa treat colorectal cancer by activating PI3K/Akt signaling pathway.
Journal of ethnopharmacology
Li XC,Wang S,Yang XX,Li TJ,Gu JX,Zhao L,Bao YR,Meng XS
Mon Jun 12 00:00:00 UTC 2023
CHD1 deletion stabilizes HIF1α to promote angiogenesis and glycolysis in prostate cancer.
Asian journal of andrology
Wang YZ,Qian YC,Yang WJ,Ye LH,Guo GD,Lv W,Huan MX,Feng XY,Wang K,Yang Z,Gao Y,Li L,Chen YL
Tue Mar 07 00:00:00 UTC 2023
Ginsenoside Rh2 inhibits CBP/p300-mediated FOXO3a acetylation and epilepsy-induced oxidative damage via the FOXO3a-KEAP1-NRF2 pathway.
European journal of pharmacology
Wu J,Wang S,Zhao W,Li M,Li S
Sun Feb 05 00:00:00 UTC 2023
Methionine alleviates heat stress-induced ferroptosis in bovine mammary epithelial cells through the Nrf2 pathway.
Ecotoxicology and environmental safety
Xu J,Wang XL,Zeng HF,Han ZY
Mon May 01 00:00:00 UTC 2023
Increased endogenous PKG I activity attenuates EGF-induced proliferation and migration of epithelial ovarian cancer via the MAPK/ERK pathway.
Cell death & disease
Lan T,Li Y,Wang Y,Wang ZC,Mu CY,Tao AB,Gong JL,Zhou Y,Xu H,Li SB,Gu B,Ma P,Luo L
Thu Jan 19 00:00:00 UTC 2023
Sufentanil combined with parecoxib sodium inhibits proliferation and metastasis of HER2-positive breast cancer cells and regulates epithelial-mesenchymal transition.
Clinical & experimental metastasis
Xu S,Li X,Li W,Ma N,Ma H,Cui J,You X,Chen X
Sat Apr 01 00:00:00 UTC 2023
Zearalenone induces oxidative stress and autophagy in goat Sertoli cells.
Ecotoxicology and environmental safety
Liu X,Xi H,Han S,Zhang H,Hu J
Wed Mar 01 00:00:00 UTC 2023
The role of 14-3-3 in the progression of vascular inflammation induced by lipopolysaccharide.
International immunopharmacology
Tan H,Li J,Jia C,Huang H,Li L,Liao B,Long Y,Nie Y,Yu F
Thu Jun 01 00:00:00 UTC 2023
A network pharmacology approach and experimental validation to investigate the anticancer mechanism and potential active targets of ethanol extract of Wei-Tong-Xin against colorectal cancer through induction of apoptosis via PI3K/AKT signaling pathway.
Journal of ethnopharmacology
Lin F,Zhang G,Yang X,Wang M,Wang R,Wan M,Wang J,Wu B,Yan T,Jia Y
Wed Mar 01 00:00:00 UTC 2023
Curcumin-dependent phenotypic transformation of microglia mediates resistance to pseudorabies-induced encephalitis.
Veterinary research
Feng L,Luo G,Li Y,Zhang C,Liu Y,Liu Y,Chen H,He D,Zhu Y,Gan L
Tue Mar 14 00:00:00 UTC 2023
Developmental growth plate cartilage formation suppressed by artificial light at night via inhibiting BMAL1-driven collagen hydroxylation.
Cell death and differentiation
Chen G,Tang Q,Yu S,Shen Y,Sun J,Peng J,Yin Y,Feng G,Lu X,Mei G,Zhang Y,Wan Q,Zhang L,Chen L
Thu Jun 01 00:00:00 UTC 2023
Mitophagy associated self-degradation of phosphorylated MAP4 guarantees the migration and proliferation responses of keratinocytes to hypoxia.
Cell death discovery
Feng Y,Li L,Zhang Q,He Y,Huang Y,Zhang J,Zhang D,Huang Y,Lei X,Hu J,Luo G
Wed May 17 00:00:00 UTC 2023
Wogonin induces ferroptosis in pancreatic cancer cells by inhibiting the Nrf2/GPX4 axis.
Frontiers in pharmacology
Liu X,Peng X,Cen S,Yang C,Ma Z,Shi X
Tue Mar 14 00:00:00 UTC 2023
ADSC-derived exosomes attenuate myocardial infarction injury by promoting miR-205-mediated cardiac angiogenesis.
Biology direct
Wang T,Li T,Niu X,Hu L,Cheng J,Guo D,Ren H,Zhao R,Ji Z,Liu P,Li Y,Guo Y
Mon Feb 27 00:00:00 UTC 2023
Design, synthesis and anti-AD effects of dual inhibitor targeting glutaminyl cyclase/GSK-3β.
European journal of medicinal chemistry
Xie Y,Chen C,Lin S,Yu X,Ye S,Chen X,Ouyang N,Xiong W,Li C,Xu C,Song G,Wu H
Wed Feb 15 00:00:00 UTC 2023
Caspase-11 contributes to pain hypersensitivity in the later phase of CFA-induced pain of mice.
Brain research
Liu M,Cao W,Qin X,Tong J,Wu X,Cheng Y
Wed Feb 15 00:00:00 UTC 2023
Neuroprotective effect of histamine H3 receptor blockade on methamphetamine-induced cognitive impairment in mice.
Pharmacology, biochemistry, and behavior
Luo H,Li X,Fan R,Ruan Y,Qian L,Shen Y,Si Z,Li L,Liu Y
Sun Jan 01 00:00:00 UTC 2023
Daucosterol Alleviates Alcohol-Induced Hepatic Injury and Inflammation through P38/NF-κB/NLRP3 Inflammasome Pathway.
Nutrients
Zhang F,Wang M,Zha Y,Zhou J,Han J,Zhang S
Sun Jan 01 00:00:00 UTC 2023
Schisandrin B Improves the Hypothermic Preservation of Celsior Solution in Human Umbilical Cord Mesenchymal Stem Cells.
Tissue engineering and regenerative medicine
Zhang Y,Wang P,Jin MX,Zhou YQ,Ye L,Zhu XJ,Li HF,Zhou M,Li Y,Li S,Liang KY,Wang Y,Gao Y,Pan MX,Zhou SQ,Peng Q
Thu Jun 01 00:00:00 UTC 2023
A phosphoglycerate mutase 1 allosteric inhibitor overcomes drug resistance to EGFR-targeted therapy via disrupting IL-6/JAK2/STAT3 signaling pathway in lung adenocarcinoma.
Drug resistance updates : reviews and commentaries in antimicrobial and anticancer chemotherapy
Liang Q,Gong M,Zou JH,Luo MY,Jiang LL,Wang C,Shen NX,Zhang MC,Xu L,Lei HM,Zhang KR,Zhang R,Zhuang G,Zhu L,Chen HZ,Zhou L,Shen Y
Mon May 01 00:00:00 UTC 2023
Sirt1 Overexpression Inhibits Fibrous Scar Formation and Improves Functional Recovery After Cerebral Ischemic Injury Through the Deacetylation of 14-3-3ζ.
Molecular neurobiology
Chen Y,Huang J,Liu J,Zhu H,Li X,Wen J,Tian M,Ren J,Zhou L,Yang Q
Tue Aug 01 00:00:00 UTC 2023
NFYAv1 is a Tumor-Promoting Transcript Associated with Poor Prognosis of Hepatocellular Carcinoma.
Medical science monitor : international medical journal of experimental and clinical research
Li Z,You C,Huang J,Zhang M
Sat Jan 21 00:00:00 UTC 2023
Allogeneic gene-edited HIV-specific CAR-T cells secreting PD-1 blocking scFv enhance specific cytotoxic activity against HIV Env+ cells invivo.
Virologica Sinica
Pan H,Yang X,Wang J,Liang H,Jiang Z,Zhao L,Wang Y,Liang Z,Shen X,Lin Q,Liang Y,Yang J,Lu P,Zhu Y,Li M,Wang P,Xu J,Lu H,Zhu H
Sat Apr 01 00:00:00 UTC 2023
Expression of group II metabotropic glutamate receptors in rat superior cervical ganglion.
Autonomic neuroscience : basic & clinical
Wei X,Zhao C,Jia X,Zhao B,Liu Y
Sun Jan 01 00:00:00 UTC 2023
LCK facilitates DNA damage repair by stabilizing RAD51 and BRCA1 in the nucleus of chemoresistant ovarian cancer.
Journal of ovarian research
Dey G,Bharti R,Braley C,Alluri R,Esakov E,Crean-Tate K,McCrae K,Joehlin-Price A,Rose PG,Lathia J,Gong Z,Reizes O
Tue Jun 27 00:00:00 UTC 2023
Thalamocortical Circuit Controls Neuropathic Pain via Up-regulation of HCN2 in the Ventral Posterolateral Thalamus.
Neuroscience bulletin
Yan Y,Zhu M,Cao X,Xu G,Shen W,Li F,Zhang J,Luo L,Zhang X,Zhang D,Liu T
Mon May 01 00:00:00 UTC 2023
Apelin-Overexpressing Neural Stem Cells in Conjunction with a Silk Fibroin Nanofiber Scaffold for the Treatment of Traumatic Brain Injury.
Stem cells and development
Li T,Tang Q,Xu J,Ye X,Chen K,Zhong J,Zhu J,Lu S,Zhu T
Fri Sep 01 00:00:00 UTC 2023
S100A1 expression is increased in spinal cord injury and promotes inflammation, oxidative stress and apoptosis of PC12 cells induced by LPS via ERK signaling.
Molecular medicine reports
Bai Y,Guo N,Xu Z,Chen Y,Zhang W,Chen Q,Bi Z
Wed Feb 01 00:00:00 UTC 2023
ANXA10 is a prognostic biomarker and suppressor of hepatocellular carcinoma: a bioinformatics analysis and experimental validation.
Scientific reports
Zhang C,Peng L,Gu H,Wang J,Wang Y,Xu Z
Sat Jan 28 00:00:00 UTC 2023
Protective efficacy of Schizandrin B on ameliorating nephrolithiasis via regulating GSK3β/Nrf2 signaling-mediated ferroptosis in vivo and in vitro.
International immunopharmacology
Dong C,Song C,He Z,Song Q,Song T,Liu J,Xiong Y,Su X,Zhou J,Yang S,Liao W
Sat Apr 01 00:00:00 UTC 2023
Pulsatilla decoction improves DSS-induced colitis via modulation of fecal-bacteria-related short-chain fatty acids and intestinal barrier integrity.
Journal of ethnopharmacology
Niu C,Hu XL,Yuan ZW,Xiao Y,Ji P,Wei YM,Hua YL
Tue Jan 10 00:00:00 UTC 2023
Exercise reduces hyperlipidemia-induced cardiac damage in apolipoprotein E-deficient mice via its effects against inflammation and oxidative stress.
Scientific reports
Pei Z,Ji J,Gao Y,Wang H,Wu Y,Yang J,Yang Q,Zhang L
Mon Jun 05 00:00:00 UTC 2023
PGC1α Degradation Suppresses Mitochondrial Biogenesis to Confer Radiation Resistance in Glioma.
Cancer research
Zhao M,Li Y,Lu C,Ding F,Xu M,Ge X,Li M,Wang Z,Yin J,Zhang J,Wang X,Ge Z,Xiao H,Xiao Y,Liu H,Liu W,Cao Y,Wang Q,You Y,Wang X,Yang K,Shi Z,Qian X
Tue Apr 04 00:00:00 UTC 2023
Decreased AGGF1 facilitates the progression of placenta accreta spectrum via mediating the P53 signaling pathway under the regulation of miR-1296-5p.
Reproductive biology
Wang R,Zhao J,Liu C,Li S,Liu W,Cao Q
Wed Mar 01 00:00:00 UTC 2023
Preventive effect of salmon sperm DNA on acute carbon tetrachloride-induced liver injury in mice through Nrf2/ARE and mitochondrial apoptosis pathway.
Food science & nutrition
Huang X,Chu X,Tian Y,Xue Y,Zhang L,Li J,Hou H,Dong P,Wang J
Wed Feb 01 00:00:00 UTC 2023
Molecular mechanism of the miR-7/BCL2L1/P53 signaling axis regulating the progression of hepatocellular carcinoma.
Annals of translational medicine
Zhang N,Wang F,Zhu L,Chang R,Mok SRS,Peixoto RD,Tang W,Chen Z
Sun Jan 15 00:00:00 UTC 2023
Glioblastoma upregulates SUMOylation of hnRNP A2/B1 to eliminate the tumor suppressor miR-204-3p, accelerating angiogenesis under hypoxia.
Cell death & disease
Guo Q,Fan Y,Wang Q,Li B,Qiu W,Qi Y,Pan Z,Zhang S,Zhao S,Yang K,Xu H,Li M,Gao Z,Xu J,Wang H,Wang S,Tang Q,Qiu J,Guo X,Deng L,Zhang P,Zhao R,Xue H,Wang C,Li G
Tue Feb 21 00:00:00 UTC 2023
Activation of PPARγ prevents ferroptosis-induced damage through Nrf2/Gpx4 signaling pathway in rat hippocampus following epilepsy.
Neuroreport
Wang MJ,Liu X,Chi S,Wang FY,Zhang L,Yao MY,Liu T,Gao J
Wed Apr 05 00:00:00 UTC 2023
Depletion of SOD2 enhances nasopharyngeal carcinoma cell radiosensitivity via ferroptosis induction modulated by DHODH inhibition.
BMC cancer
Amos A,Jiang N,Zong D,Gu J,Zhou J,Yin L,He X,Xu Y,Wu L
Fri Feb 03 00:00:00 UTC 2023
Exosomes from Human Umbilical Cord Mesenchymal Stem Cells Facilitates Injured Endometrial Restoring in Early Repair Period through miR-202-3p Mediating Formation of ECM.
Stem cell reviews and reports
Wang S,Liu T,Nan N,Lu C,Liang M,Wang S,Wang H,He B,Chen X,Xu X,Zheng Y
Tue Aug 01 00:00:00 UTC 2023
Adjuvant therapy with Huatan Sanjie Granules improves the prognosis of patients with primary liver cancer: a cohort study and the investigation of its mechanism of action based on network pharmacology.
Frontiers in pharmacology
Yuan J,Abdurahman A,Cui N,Hao T,Zou J,Liu L,Wu Y
Mon Jun 19 00:00:00 UTC 2023
Decreasing GDF15 Promotes Inflammatory Signals and Neutrophil Infiltration in Psoriasis Models.
The Journal of investigative dermatology
Zhang J,He L,Wang Z,Shao S,Qiao P,Zhang J,Zhang K,Li C,Zhang Y,Wang G,Li M
Wed Mar 01 00:00:00 UTC 2023
Supplementing a specific synbiotic suppressed the incidence of AOM/DSS-induced colorectal cancer in mice.
iScience
Wu H,Wu Z,Qiu Y,Zhao F,Liao M,Zhong Z,Chen J,Zeng Y,Liu R
Fri Jun 16 00:00:00 UTC 2023
Two-Pore-Domain Potassium Channel TREK-1 Mediates Pulmonary Fibrosis through Macrophage M2 Polarization and by Direct Promotion of Fibroblast Differentiation.
Biomedicines
Zhang Y,Fu J,Han Y,Feng D,Yue S,Zhou Y,Luo Z
Wed Apr 26 00:00:00 UTC 2023
MC1R Peptide Agonist Self-Assembles into a Hydrogel That Promotes Skin Pigmentation for Treating Vitiligo.
ACS nano
Zhu C,Li T,Wang Z,Li Z,Wei J,Han H,Yuan D,Cai M,Shi J
Tue May 09 00:00:00 UTC 2023
VprBP/DCAF1 triggers melanomagenic gene silencing through histone H2A phosphorylation.
Research square
Shin Y,Kim S,Liang G,Ulmer TS,An W
Wed Jul 12 00:00:00 UTC 2023
Polystyrene Microplastics Induce Oxidative Stress in Mouse Hepatocytes in Relation to Their Size.
International journal of molecular sciences
Zou H,Qu H,Bian Y,Sun J,Wang T,Ma Y,Yuan Y,Gu J,Bian J,Liu Z
Mon Apr 17 00:00:00 UTC 2023
Naringin regulates endoplasmic reticulum stress and mitophagy through the ATF3/PINK1 signaling axis to alleviate pulmonary fibrosis.
Naunyn-Schmiedeberg's archives of pharmacology
Wei Y,Sun L,Liu C,Li L
Thu Jun 01 00:00:00 UTC 2023
Glycoside combinations of Buyang Huanwu decoction ameliorate atherosclerosis via STAT3, HIF-1, and VEGF.
Naunyn-Schmiedeberg's archives of pharmacology
Yan F,Ding H,Sun Z,Liu J,Li J,Zhou D,Zhang W
Thu Jun 01 00:00:00 UTC 2023
Scutellarin Alleviates Ischemic Brain Injury in the Acute Phase by Affecting the Activity of Neurotransmitters in Neurons.
Molecules (Basel, Switzerland)
Wang C,Liu Y,Liu X,Zhang Y,Yan X,Deng X,Shi J
Tue Apr 04 00:00:00 UTC 2023
Huntingtin-associated protein 1 is a potential tumor suppressor for gastric cancer.
Molecular biology reports
Qu YM,Chen A,Zhao X,Wang Z,Guo D,Shao SL,Tao YY,Li QJ,Wang MY,Ma WS
Wed Feb 01 00:00:00 UTC 2023
Epitranscriptic regulation of HRAS by N6-methyladenosine drives tumor progression.
Proceedings of the National Academy of Sciences of the United States of America
Pan Y,Gu Y,Liu T,Zhang Q,Yang F,Duan L,Cheng S,Zhu X,Xi Y,Chang X,Ye Q,Gao S
Tue Apr 04 00:00:00 UTC 2023
6-(7-Nitro-2,1,3-benzoxadiazol-4-ylthio) Hexanol Inhibits Proliferation and Induces Apoptosis of Endometriosis by Regulating Glutathione S-Transferase Mu Class 4.
Reproductive sciences (Thousand Oaks, Calif.)
Liu W,Cheng L,Du Y,Liu X,Ma J,Yan L
Sun Oct 01 00:00:00 UTC 2023
Metabolic genes, a potential predictor of prognosis and immunogenicity of clear cell renal cell carcinoma.
American journal of cancer research
Ji CJ,Wei XY,Yao LY,Wang YC,Cong R,Song NH
Sun May 14 00:00:00 UTC 2023
Glucose Starvation Inhibits Ferroptosis by Activating the LKB1/AMPK Signaling Pathway and Promotes the High Speed Linear Motility of Dairy Goat Sperm.
Animals : an open access journal from MDPI
Li Y,Zhang G,Wen F,Xian M,Guo S,Zhang X,Feng X,Hu Z,Hu J
Sun Apr 23 00:00:00 UTC 2023
Inhibition of STEAP1 ameliorates inflammation and ferroptosis of acute lung injury caused by sepsis in LPS-induced human pulmonary microvascular endothelial cells.
Molecular biology reports
Zou X,Liu C,Huang Z,Xiang S,Li K,Yuan Y,Hao Y,Zhou F
Sat Jul 01 00:00:00 UTC 2023
SETD7 promotes lateral plate mesoderm formation by modulating the Wnt/β-catenin signaling pathway.
iScience
Wang D,Li Y,Xu C,Wang H,Huang X,Jin X,Ren S,Gao J,Tong J,Liu J,Zhou J,Shi L
Fri Jun 16 00:00:00 UTC 2023
Acupuncture regulates the apoptosis of ovarian granulosa cells in polycystic ovarian syndrome-related abnormal follicular development through LncMEG3-mediated inhibition of miR-21-3p.
Biological research
Chen X,He H,Long B,Wei B,Yang P,Huang X,Wang Q,Lin J,Tang H
Mon Jun 12 00:00:00 UTC 2023
Effect of stem cell conditional medium-loading adhesive hydrogel on TGF-β1-induced endometrial stromal cell fibrosis.
Frontiers in bioengineering and biotechnology
Zhu Y,Wang T,Bao MJ,Qu XH,Li ZM
Tue May 23 00:00:00 UTC 2023
Anti-tumor effect of infant-derived Enterococcus via the inhibition of proliferation and inflammation as well as the promotion of apoptosis.
Food & function
Yang Q,He Y,Tian L,Zhang Z,Qiu L,Tao X,Wei H
Tue Feb 21 00:00:00 UTC 2023
Shikonin suppresses small cell lung cancer growth via inducing ATF3-mediated ferroptosis to promote ROS accumulation.
Chemico-biological interactions
Qian X,Zhu L,Xu M,Liu H,Yu X,Shao Q,Qin J
Fri Sep 01 00:00:00 UTC 2023
Semaglutide inhibits ischemia/reperfusion-induced cardiomyocyte apoptosis through activating PKG/PKCε/ERK1/2 pathway.
Biochemical and biophysical research communications
Zhu Q,Luo Y,Wen Y,Wang D,Li J,Fan Z
Sun Mar 05 00:00:00 UTC 2023
Atractylenolide III suppresses senescence-associated secretome via inhibiting cGAS/NF-κB pathway in hepatic stellate cells.
Clinical and experimental pharmacology & physiology
Wu H,Wu L,Xiao L,Gu Y,Liu H,Zhang L,Zhang M,Qi L
Sat Apr 01 00:00:00 UTC 2023
Exendin-4 alleviates myocardial ischemia reperfusion injury by enhancing autophagy through promoting nuclear translocation of TFEB.
Experimental cell research
He W,Tong G,Fan H,Zhen C,Zeng L,Xue L,Chen J,Sun Z,He P
Wed Feb 15 00:00:00 UTC 2023
Zebrafish Galectin 3 binding protein is the target antigen of the microglial 4C4 monoclonal antibody.
Developmental dynamics : an official publication of the American Association of Anatomists
Rovira M,Miserocchi M,Montanari A,Hammou L,Chomette L,Pozo J,Imbault V,Bisteau X,Wittamer V
Wed Mar 01 00:00:00 UTC 2023
NPAS2 promotes aerobic glycolysis and tumor growth in prostate cancer through HIF-1A signaling.
BMC cancer
Ma S,Chen Y,Quan P,Zhang J,Han S,Wang G,Qi R,Zhang X,Wang F,Yuan J,Yang X,Jia W,Qin W
Tue Mar 28 00:00:00 UTC 2023
Silencing of OGDHL promotes liver cancer metastasis by enhancing hypoxia inducible factor 1 α protein stability.
Cancer science
Dai W,Li Y,Sun W,Ji M,Bao R,Chen J,Xu S,Dai Y,Chen Y,Liu W,Ge C,Sun W,Mo W,Guo C,Xu X
Sat Apr 01 00:00:00 UTC 2023
Afatinib combined with anti-PD1 enhances immunotherapy of hepatocellular carcinoma via ERBB2/STAT3/PD-L1 signaling.
Frontiers in oncology
Yu C,Zhang X,Wang M,Xu G,Zhao S,Feng Y,Pan C,Yang W,Zhou J,Shang L,Ma Y
Mon Jun 19 00:00:00 UTC 2023
P4HA1 activates HMGCS1 to promote nasopharyngeal carcinoma ferroptosis resistance and progression.
Cellular signalling
Zhou R,Qiu L,Zhou L,Geng R,Yang S,Wu J
Mon May 01 00:00:00 UTC 2023
Irisin Ameliorates PM2.5-Induced Acute Lung Injury by Regulation of Autophagy Through AMPK/mTOR Pathway.
Journal of inflammation research
Ma J,Han Z,Jiao R,Yuan G,Ma C,Yan X,Meng A
Tue Mar 21 00:00:00 UTC 2023
Aescin can alleviate NAFLD through Keap1-Nrf2 by activating antioxidant and autophagy.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yu H,Yan S,Jin M,Wei Y,Zhao L,Cheng J,Ding L,Feng H
Mon May 01 00:00:00 UTC 2023
Constitutively activated AMPKα1 protects against skeletal aging in mice by promoting bone-derived IGF-1 secretion.
Cell proliferation
Yang Y,Yuan K,Liu Y,Wang Q,Lin Y,Yang S,Huang K,Kan T,Zhang Y,Xu M,Yu Z,Fan Q,Wang Y,Li H,Tang T
Sun Oct 01 00:00:00 UTC 2023
Construction and Rescue of a DNA-Launched DENV2 Infectious Clone.
Viruses
Holliday M,Corliss L,Lennemann NJ
Wed Jan 18 00:00:00 UTC 2023
Protective effects of berberine against β-amyloid-induced neurotoxicity in HT22 cells via the Nrf2/HO-1 pathway.
Bioorganic chemistry
Zhang RL,Lei BX,Wu GY,Wang YY,Huang QH
Sat Apr 01 00:00:00 UTC 2023
The histone H2B ubiquitination regulator Wac is essential for plasma cell differentiation.
FEBS letters
Li Y,Ruan GX,Chen W,Huang H,Zhang R,Wang J,Ouyang Y,Zhu Z,Meng L,Wang R,Huo J,Xu S,Ou X
Sat Jul 01 00:00:00 UTC 2023
Nervonic acid improves liver inflammation in a mouse model of Parkinson's disease by inhibiting proinflammatory signaling pathways and regulating metabolic pathways.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang X,Liang T,Mao Y,Li Z,Li X,Zhu X,Cao F,Zhang J
Tue Aug 01 00:00:00 UTC 2023
SEL1L-HRD1 endoplasmic reticulum-associated degradation controls STING-mediated innate immunity by limiting the size of the activable STING pool.
Nature cell biology
Ji Y,Luo Y,Wu Y,Sun Y,Zhao L,Xue Z,Sun M,Wei X,He Z,Wu SA,Lin LL,Lu Y,Chang L,Chen F,Chen S,Qian W,Xu X,Chen S,Pan D,Zhou Z,Xia S,Hu CA,Liang T,Qi L
Mon May 01 00:00:00 UTC 2023
Preoperative Pain Facilitates Postoperative Cognitive Dysfunction via Periaqueductal Gray Matter-Dorsal Raphe Circuit.
Neuroscience
Deng H,Wu Y,Gao P,Kong D,Pan C,Xu S,Tang D,Jiao Y,Wen D,Yu W
Tue Aug 01 00:00:00 UTC 2023
SATB1 regulates 3D genome architecture in T cells by constraining chromatin interactions surrounding CTCF-binding sites.
Cell reports
Wang B,Ji L,Bian Q
Tue Apr 25 00:00:00 UTC 2023
LncRNA FRMD6-AS1 promotes hepatocellular carcinoma cell migration and stemness by regulating SENP1/HIF-1α axis.
Pathology, research and practice
Sun W,Lei X,Lu Q,Wu Q,Ma Q,Huang D,Zhang Y
Wed Mar 01 00:00:00 UTC 2023
The mechanism of monobutyl phthalate -induced ferroptosis via TNF/IL6/STAT3 signal pathway in TM-3 cells.
The Journal of toxicological sciences
Guo X,Hao Y,Ma H,Li H,Li L,Yan F,Huang J,Li L
Tue May 02 00:00:00 UTC 2023
BNIP3-mediated autophagy via the mTOR/ULK1 pathway induces primordial follicle loss after ovarian tissue transplantation.
Journal of assisted reproduction and genetics
Liu F,Li M
Wed Mar 01 00:00:00 UTC 2023
Targeting hexokinase 1 alleviates NLRP3-mediated inflammation in apical periodontitis: A laboratory investigation.
International endodontic journal
Qian Y,Chen D,Zhu Y,Wu J,Wang Y,Yang W
Thu Jun 01 00:00:00 UTC 2023
Irisin attenuates fine particulate matter induced acute lung injury by regulating Nod2/NF-κB signaling pathway.
Immunobiology
Jiao R,Han Z,Ma J,Wu S,Wang Z,Zhou G,Liu X,Li J,Yan X,Meng A
Mon May 01 00:00:00 UTC 2023
TGF-β3 Protects Neurons Against Intermittent Hypoxia-Induced Oxidative Stress and Apoptosis Through Activation of the Nrf-2/KEAP1/HO-1 Pathway via Binding to TGF-βRI.
Neurochemical research
Huang Y,Liu Z,Wang X,Li Y,Liu L,Li B
Fri Sep 01 00:00:00 UTC 2023
Diosmetin alleviates acute lung injury caused by lipopolysaccharide by targeting barrier function.
Inflammopharmacology
Xia J,Li J,Deng M,Yin F,Liu J,Wang J
Tue Aug 01 00:00:00 UTC 2023
Ring domains are essential for GATOR2-dependent mTORC1 activation.
Molecular cell
Jiang C,Dai X,He S,Zhou H,Fang L,Guo J,Liu S,Zhang T,Pan W,Yu H,Fu T,Li D,Inuzuka H,Wang P,Xiao J,Wei W
Thu Jan 05 00:00:00 UTC 2023
Protective effects of pentoxifylline against chlorine-induced acute lung injury in rats.
BMC pharmacology & toxicology
Liu MM,Liu JZ,Zhao CQ,Guo P,Wang Z,Wu H,Yu W,Liu R,Hai CX,Zhang XD
Mon Feb 27 00:00:00 UTC 2023
The RNA-binding protein hnRNP F is required for the germinal center B cell response.
Nature communications
Huang H,Li Y,Zhang G,Ruan GX,Zhu Z,Chen W,Zou J,Zhang R,Wang J,Ouyang Y,Xu S,Ou X
Thu Mar 30 00:00:00 UTC 2023
Proteomic analysis of DZIP3 interactome and its role in proliferation and metastasis in gastric cancer cells.
Experimental cell research
Qiu R,Zhao S,Lu C,Xu Z,Shu E,Weng Q,Chen W,Fang S,Chen W,Zheng L,Zhao Z,Yang Y,Ji J
Sat Apr 01 00:00:00 UTC 2023
The promoting effect and mechanism of Nrf2 on cell metastasis in cervical cancer.
Journal of translational medicine
Zhang M,Hong X,Ma N,Wei Z,Ci X,Zhang S
Tue Jul 04 00:00:00 UTC 2023
TREM2 Inhibits Tau Hyperphosphorylation and Neuronal Apoptosis via the PI3K/Akt/GSK-3β Signaling Pathway In vivo and In vitro.
Molecular neurobiology
Peng X,Guo H,Zhang X,Yang Z,Ruganzu JB,Yang Z,Wu X,Bi W,Ji S,Yang W
Mon May 01 00:00:00 UTC 2023
Mesenchymal stem cell-derived extracellular vesicles carrying miR-99b-3p restrain microglial activation and neuropathic pain by stimulating autophagy.
International immunopharmacology
Gao X,Gao LF,Kong XQ,Zhang YN,Jia S,Meng CY
Wed Feb 01 00:00:00 UTC 2023
Synergistic effects of combined treatment of 1,2-dichloroethane and high-dose ethanol on liver damage in mice and the related mechanisms.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Yang J,Zhang L,Wang T,Zhang J,Li M,Jin X,Tan X,Wang G,Zhao F,Jin Y
Thu Jun 01 00:00:00 UTC 2023
Rho A/ROCK1 signaling-mediated metabolic reprogramming of valvular interstitial cells toward Warburg effect accelerates aortic valve calcification via AMPK/RUNX2 axis.
Cell death & disease
Liu H,Yin H,Wang Z,Yuan Q,Xu F,Chen Y,Li C
Sat Feb 11 00:00:00 UTC 2023
Supplemental Clostridium butyricum modulates lipid metabolism by reshaping the gut microbiota composition and bile acid profile in IUGR suckling piglets.
Journal of animal science and biotechnology
Zhang X,Yun Y,Lai Z,Ji S,Yu G,Xie Z,Zhang H,Zhong X,Wang T,Zhang L
Mon Mar 13 00:00:00 UTC 2023
A novel 2-iminobenzimidazole compound, XYA1353, displays in vitro and in vivo anti-myeloma activity via targeting NF-κB signaling.
Molecular and cellular biochemistry
Gao J,Zhou J,Zhang M,Zhang Y,Zeng Y,Li S,Xu K,Yao R
Fri May 19 00:00:00 UTC 2023
Mitochondria-Targeted Nanosystem Enhances Radio-Radiodynamic-Chemodynamic Therapy on Triple Negative Breast Cancer.
ACS applied materials & interfaces
Li J,You Z,Zhai S,Zhao J,Lu K
Wed May 10 00:00:00 UTC 2023
Sphingosine-1-Phosphate Receptor 4 Attenuates Neutrophilic Airway Inflammation in Experimental Asthma via Repressing Proinflammatory Macrophage Activation.
International journal of biological sciences
Wang S,Tian Z,Lu Y,Huang Z,Fan Y,Li B,Zheng H,Wu X,Wang M,Zhao J,Xie J
Mon Apr 17 00:00:00 UTC 2023
NCAPG is transcriptionally regulated by CBX3 and activates the Wnt/β-catenin signaling pathway to promote proliferation and the cell cycle and inhibit apoptosis in colorectal cancer.
Journal of gastrointestinal oncology
Yang H,Pu L,Li R,Zhu R
Sat Apr 29 00:00:00 UTC 2023
Diet Control and Swimming Exercise Ameliorate HFD-Induced Cognitive Impairment Related to the SIRT1-NF-κB/PGC-1α Pathways in ApoE-/- Mice.
Neural plasticity
Wei W,Lin Z,Xu P,Lv X,Lin L,Li Y,Zhou Y,Lu T,Xue X
Mon Apr 03 00:00:00 UTC 2023
Danlou tablet inhibits high-glucose-induced cardiomyocyte apoptosis via the miR-34a-SIRT1 axis.
Heliyon
Chen R,Chen H,Yang Z,Zhu L,Bei Y,Chen W,Qiu Y
Wed Mar 01 00:00:00 UTC 2023
Knockdown of Brg1 reduced mucus secretion in HDM stimulated airway inflammation.
Molecular immunology
Xu M,Hu J,Yang L,Gen G,Fu Z,Luo Z,Zou W
Sun Jan 01 00:00:00 UTC 2023
Exosomes derived from BMSCs ameliorate cyclophosphamide-induced testosterone deficiency by enhancing the autophagy of Leydig cells via the AMPK-mTOR signaling pathway.
Asian journal of andrology
Liang HY,Peng F,Pan MJ,Liao SL,Wei C,Wei GY,Xie X,Xue KY,Chen MK,Yang JK,Guo WB,Liu CD,Zhou QZ
Fri Jul 21 00:00:00 UTC 2023
The antiviral activity of tripartite motif protein 38 in hepatitis B virus replication and gene expression and its association with treatment responses during PEG-IFN-α antiviral therapy.
Virology
Luo H,Hu X,Li Y,Lei D,Tan G,Zeng Y,Qin B
Wed Feb 01 00:00:00 UTC 2023
Fluvoxamine alleviates bleomycin-induced lung fibrosis via regulating the cGAS-STING pathway.
Pharmacological research
Xie X,Wu X,Zhao D,Liu Y,Du Q,Li Y,Xu Y,Li Y,Qiu Y,Yang Y
Sun Jan 01 00:00:00 UTC 2023
Macrophage migration inhibitory factor facilitates astrocytic production of the CCL2 chemokine following spinal cord injury.
Neural regeneration research
Zhang H,Hu YM,Wang YJ,Zhou Y,Zhu ZJ,Chen MH,Wang YJ,Xu H,Wang YH
Tue Aug 01 00:00:00 UTC 2023
Cartilage Damage Pathological Characteristics of Diabetic Neuropathic Osteoarthropathy.
Analytical cellular pathology (Amsterdam)
Liu PL,Diao JY,Wang Q,Liu H,Zhang Y,Liang JQ,Zhang F,Liang XJ,Zhao HM
Fri May 19 00:00:00 UTC 2023
Transcytosable Peptide-Paclitaxel Prodrug Nanoparticle for Targeted Treatment of Triple-Negative Breast Cancer.
International journal of molecular sciences
Wang L,Zhao C,Lu L,Jiang H,Wang F,Zhang X
Tue Feb 28 00:00:00 UTC 2023
Gut Microbiota Taxon-Dependent Transformation of Microglial M1/M2 Phenotypes Underlying Mechanisms of Spatial Learning and Memory Impairment after Chronic Methamphetamine Exposure.
Microbiology spectrum
Wu Y,Dong Z,Jiang X,Qu L,Zhou W,Sun X,Hou J,Xu H,Cheng M
Thu Jun 15 00:00:00 UTC 2023
Diosgenin attenuates nonalcoholic hepatic steatosis through the hepatic FXR-SHP-SREBP1C/PPARα/CD36 pathway.
European journal of pharmacology
Chen S,Sun S,Feng Y,Li X,Yin G,Liang P,Yu W,Meng D,Zhang X,Liu H,Zhang F
Sat Aug 05 00:00:00 UTC 2023
Comprehensive analysis of lncRNA-mRNA co-expression networks in HPV-driven cervical cancer reveals the pivotal function of LINC00511-PGK1 in tumorigenesis.
Computers in biology and medicine
Xin X,Jia-Yin Y,Jun-Yang H,Rui W,Xiong-Ri K,Long-Rui D,Liu J,Jue-Yu Z
Thu Jun 01 00:00:00 UTC 2023
Silencing TRAIP suppresses cell proliferation and migration/invasion of triple negative breast cancer via RB-E2F signaling and EMT.
Cancer gene therapy
Zheng Y,Jia H,Wang P,Liu L,Chen Z,Xing X,Wang J,Tan X,Wang C
Sun Jan 01 00:00:00 UTC 2023
Anti-Postmenopausal osteoporosis effects of Isopsoralen: A bioinformatics-integrated experimental study.
Phytotherapy research : PTR
Li H,Wang C,Jin Y,Cai Y,Yao J,Meng Q,Wu J,Wang H,Sun H,Liu M
Sun Jan 01 00:00:00 UTC 2023
Naringin protects against inflammation and apoptosis induced by intestinal ischemia-reperfusion injury through deactivation of cGAS-STING signaling pathway.
Phytotherapy research : PTR
Gu L,Wang F,Wang Y,Sun D,Sun Y,Tian T,Meng Q,Yin L,Xu L,Lu X,Peng J,Lin Y,Sun P
Tue Aug 01 00:00:00 UTC 2023
Apurinic/apyrimidinic endonuclease 1 regulates palmitic acid-mediated apoptosis in cardiomyocytes via endoplasmic reticulum stress.
Biochemical and biophysical research communications
Fan J,Liu M,Li X,Gao S,Wang Y,Li A,Chen L,Zhou D,Chen H,Xu Z,Wu Z,Wu K
Sun Apr 02 00:00:00 UTC 2023
Two Resveratrol Oligomers Inhibit Cathepsin L Activity to Suppress SARS-CoV-2 Entry.
Journal of agricultural and food chemistry
Wang C,Ye X,Ding C,Zhou M,Li W,Wang Y,You Q,Zong S,Peng Q,Duanmu D,Chen H,Sun B,Qiao J
Wed Apr 12 00:00:00 UTC 2023
Endothelial Arid1a deletion disrupts the balance among angiogenesis, neurogenesis and gliogenesis in the developing brain.
Cell proliferation
Wang Y,Su L,Wang W,Zhao J,Wang Y,Li S,Liu Y,Chai R,Li X,Teng Z,Liu C,Hu B,Ji F,Jiao J
Mon May 01 00:00:00 UTC 2023
Study on the pharmacological mechanisms of sodium-glucose co-transporter 2 inhibitors in obesity-related atrial fibrillation based on network pharmacology and experimental verification.
Annals of translational medicine
Zhao Q,Wang P,Zhao M,Hong J,Zhang N,Lin K,Li Y
Fri Apr 28 00:00:00 UTC 2023
Synthesis of Novel Pentacyclic Triterpenoid Derivatives that Induce Apoptosis in Cancer Cells through a ROS-dependent, Mitochondrial-Mediated Pathway.
Molecular pharmaceutics
Cheng B,Chu X,Liu R,Ma X,Wang M,Zhang J,Jiao P,Gao Q,Ma W,Zhang Y,Zhao C,Zhou D,Xiao S
Mon Jan 02 00:00:00 UTC 2023
PbO nanoparticles increase the expression of ICAM-1 and VCAM-1 by increasing reactive oxygen species production in choroid plexus.
Environmental science and pollution research international
Wang W,Li S,Wang X,Wang J,Zhang Y
Wed Mar 01 00:00:00 UTC 2023
LncRNA SOCS2-AS1 promotes the progression of papillary thyroid cancer by destabilizing p53 protein.
Biochemical and biophysical research communications
Zhang X,Zhang X,Yang G,Wan L,Yin F,Li H,Yin D
Sun Aug 20 00:00:00 UTC 2023
Exosomes Derived from AT2R-Overexpressing BMSC Prevent Restenosis After Carotid Artery Injury by Attenuating the Injury-Induced Neointimal Hyperplasia.
Journal of cardiovascular translational research
Zou X,Liao Y,Liu Z,Xu X,Sun W,Qin H,Wang H,Liu J,Jing T
Wed Feb 01 00:00:00 UTC 2023
Title: Bioinformatic Identification of Genes Involved in Diabetic Nephropathy Fibrosis and their Clinical Relevance.
Biochemical genetics
Bai Y,Ma L,Deng D,Tian D,Liu W,Diao Z
Tue Aug 01 00:00:00 UTC 2023
Polyphyllin D punctures hypertrophic lysosomes to reverse drug resistance of hepatocellular carcinoma by targeting acid sphingomyelinase.
Molecular therapy : the journal of the American Society of Gene Therapy
Wang Y,Chen YY,Gao GB,Zheng YH,Yu NN,Ouyang L,Gao X,Li N,Wen SY,Huang S,Zhao Q,Liu L,Cao M,Zhang S,Zhang J,He QY
Wed Jul 05 00:00:00 UTC 2023
Ginsenoside Rg1 protects cardiac mitochondrial function via targeting GSTP1 to block S-glutathionylation of optic atrophy 1.
Free radical biology & medicine
Zhu H,Yan C,Yao P,Li P,Li Y,Yang H
Tue Aug 01 00:00:00 UTC 2023
Electroacupuncture alleviates early brain injury via modulating microglia polarization and suppressing neuroinflammation in a rat model of subarachnoid hemorrhage.
Heliyon
Wang Y,Yang X,Cao Y,Li X,Xu R,Yan J,Guo Z,Sun S,Sun X,Wu Y
Wed Mar 01 00:00:00 UTC 2023
YY1 lactylation in microglia promotes angiogenesis through transcription activation-mediated upregulation of FGF2.
Genome biology
Wang X,Fan W,Li N,Ma Y,Yao M,Wang G,He S,Li W,Tan J,Lu Q,Hou S
Fri Apr 21 00:00:00 UTC 2023
mTOR/ULK1 signaling axis mediated ultrashort wave regulation of autophagy to alleviate diabetic kidney disease.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University
Yan Y,Yu S,Fan Y
Wed Nov 01 00:00:00 UTC 2023
Schisandrin B promotes senescence of activated hepatic stellate cell via NCOA4-mediated ferritinophagy.
Pharmaceutical biology
Ma M,Wei N,Yang J,Ding T,Song A,Chen L,Zheng S,Jin H
Fri Dec 01 00:00:00 UTC 2023
Farnesoid X Receptor (FXR) Regulates mTORC1 Signaling and Autophagy by Inhibiting SESN2 Expression.
Molecular nutrition & food research
Wang G,Chen L,Qin S,Geng H,Xia C,Zheng Y,Lei X,Zhang J,Wu S,Yao J,Deng L
Wed Mar 01 00:00:00 UTC 2023
Farnesoid X Receptor (FXR) Regulates mTORC1 Signaling and Autophagy by Inhibiting SESN2 Expression.
Molecular nutrition & food research
Wang G,Chen L,Qin S,Geng H,Xia C,Zheng Y,Lei X,Zhang J,Wu S,Yao J,Deng L
Wed Mar 01 00:00:00 UTC 2023
MiR-29c Inhibits TNF-α-Induced ROS Production and Apoptosis in Mouse Hippocampal HT22 Cell Line.
Neurochemical research
Li B,Lu Y,Wang R,Xu T,Lei X,Jin H,Gao X,Xie Y,Liu X,Zeng J
Wed Feb 01 00:00:00 UTC 2023
Parecoxib attenuates inflammation injury in septic H9c2 cells by regulating the MAPK signaling pathway.
Experimental and therapeutic medicine
Qian X,Xiong S,Chen Q,Zhang J,Xie J
Sat Apr 01 00:00:00 UTC 2023
Aprepitant inhibits the progression of esophageal squamous cancer by blocking the truncated neurokinin‑1 receptor.
Oncology reports
Zheng Y,Sang M,Liu F,Gu L,Li J,Wu Y,Shan B
Sat Jul 01 00:00:00 UTC 2023
Selenium supplementation enhanced the expression of selenoproteins in hippocampus and played a neuroprotective role in LPS-induced neuroinflammation.
International journal of biological macromolecules
Liang X,Xue Z,Zheng Y,Li S,Zhou L,Cao L,Zou Y
Sat Apr 15 00:00:00 UTC 2023
DNA replication initiation factor RECQ4 possesses a role in antagonizing DNA replication initiation.
Nature communications
Xu X,Chang CW,Li M,Omabe K,Le N,Chen YH,Liang F,Liu Y
Sat Mar 04 00:00:00 UTC 2023
LncRNA UCA1 promoted cisplatin resistance in lung adenocarcinoma with HO1 targets NRF2/HO1 pathway.
Journal of cancer research and clinical oncology
Shi W,Ling L,Li C,Wu R,Zhang M,Shao F,Wang Y
Wed Mar 01 00:00:00 UTC 2023
Role of SESTRIN2/AMPK/ULK1 pathway activation and lysosomes dysfunction in NaAsO2-induced liver injury under oxidative stress.
Ecotoxicology and environmental safety
Bi D,Zheng D,Shi M,Hu Q,Wang H,Zhi H,Lou D,Zhang A,Hu Y
Sat Apr 01 00:00:00 UTC 2023
4-Octyl itaconate attenuates LPS-induced acute kidney injury by activating Nrf2 and inhibiting STAT3 signaling.
Molecular medicine (Cambridge, Mass.)
Xu L,Cai J,Li C,Yang M,Duan T,Zhao Q,Xi Y,Sun L,He L,Tang C,Sun L
Mon Apr 24 00:00:00 UTC 2023
Pine pollen extract alleviates ethanol-induced oxidative stress and apoptosis in HepG2 cells via MAPK signaling.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Ma D,Wang Z,He Z,Wang Z,Chen Q,Qin F,Zeng M,Chen J
Sun Jan 01 00:00:00 UTC 2023
Dose-Dependent Effects in Plasma Oncotherapy: Critical In Vivo Immune Responses Missed by In Vitro Studies.
Biomolecules
He Y,Gong F,Jin T,Liu Q,Fang H,Chen Y,Wang G,Chu PK,Wu Z,Ostrikov KK
Fri Apr 21 00:00:00 UTC 2023
SlMYB99-mediated auxin and abscisic acid antagonistically regulate ascorbic acids biosynthesis in tomato.
The New phytologist
Xu X,Huang B,Fang X,Zhang Q,Qi T,Gong M,Zheng X,Wu M,Jian Y,Deng J,Cheng Y,Li Z,Deng W
Tue Aug 01 00:00:00 UTC 2023
The main e-cigarette component vegetable glycerin enhances neutrophil migration and fibrosis in endotoxin-induced lung injury via p38 MAPK activation.
Respiratory research
Su VY,Chen WC,Yu WK,Wu HH,Chen H,Yang KY
Tue Jan 10 00:00:00 UTC 2023
The circadian rhythm regulates branched-chain amino acids metabolism in fast muscle of Chinese perch (Siniperca chuatsi) during short-term fasting by Clock-KLF15-Bcat2 pathway.
The British journal of nutrition
Zhu X,Liu J,Cai M,Bao L,Pan Y,Wu P,Chu W,Zhang J
Mon Aug 28 00:00:00 UTC 2023
The circadian rhythm gene Bmal1 ameliorates acute deoxynivalenol-induced liver damage.
Archives of toxicology
Yang LN,Xu S,Tang M,Zhou X,Liao Y,Nüssler AK,Liu L,Yang W
Wed Mar 01 00:00:00 UTC 2023
Neobavaisoflavone induces pyroptosis of liver cancer cells via Tom20 sensing the activated ROS signal.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li Y,Zhao R,Xiu Z,Yang X,Zhu Y,Han J,Li S,Li Y,Sun L,Li X,Jin N,Li Y
Tue Jul 25 00:00:00 UTC 2023
1α,25(OH)2D3 Promotes the Autophagy of Porcine Ovarian Granulosa Cells as a Protective Mechanism against ROS through the BNIP3/PINK1 Pathway.
International journal of molecular sciences
Wang S,Yao Q,Zhao F,Cui W,Price CA,Wang Y,Lv J,Tang H,Jiang Z
Wed Feb 22 00:00:00 UTC 2023
Sirt3-dependent regulation of mitochondrial oxidative stress and apoptosis contributes to the dysfunction of pancreatic islets after severe burns.
Free radical biology & medicine
Liu X,Xie X,Li D,Liu Z,Zhang B,Zang Y,Yuan H,Shen C
Wed Mar 01 00:00:00 UTC 2023
Role of FLCN Phosphorylation in Insulin-Mediated mTORC1 Activation and Tumorigenesis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wang G,Chen L,Lei X,Qin S,Geng H,Zheng Y,Xia C,Yao J,Meng T,Deng L
Thu Jun 01 00:00:00 UTC 2023
Role of FLCN Phosphorylation in Insulin-Mediated mTORC1 Activation and Tumorigenesis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wang G,Chen L,Lei X,Qin S,Geng H,Zheng Y,Xia C,Yao J,Meng T,Deng L
Thu Jun 01 00:00:00 UTC 2023
Chlorogenic acid combined with epigallocatechin-3-gallate mitigates D-galactose-induced gut aging in mice.
Food & function
Wei R,Su Z,Mackenzie GG
Mon Mar 20 00:00:00 UTC 2023
Inhibition of Checkpoint Kinase 1 (CHK1) Upregulates Interferon Regulatory Factor 1 (IRF1) to Promote Apoptosis and Activate Anti-Tumor Immunity via MICA in Hepatocellular Carcinoma (HCC).
Cancers
Li X,Huang J,Wu Q,Du Q,Wang Y,Huang Y,Cai X,Geller DA,Yan Y
Mon Jan 30 00:00:00 UTC 2023
Microcystin‑leucine arginine promotes colorectal cancer cell proliferation by activating the PI3K/Akt/Wnt/β‑catenin pathway.
Oncology reports
Tang Y,Yi X,Zhang X,Liu B,Lu Y,Pan Z,Yu T,Feng W
Sun Jan 01 00:00:00 UTC 2023
PLD2 deletion alleviates disruption of tight junctions in sepsis-induced ALI by regulating PA/STAT3 phosphorylation pathway.
International immunopharmacology
Qian T,Qi B,Fei Y,Li J,Luo L,Lv B,Song Y,Sheng S,Xiao W,Huang X,Wang X
Sun Jan 01 00:00:00 UTC 2023
Application of neural network-based image analysis to detect sister chromatid cohesion defects.
Scientific reports
Ikemoto D,Taniguchi T,Hirota K,Nishikawa K,Okubo K,Abe T
Mon Feb 06 00:00:00 UTC 2023
An in vivo and in vitro model on the protective effect of cilnidipine on contrast-induced nephropathy via regulation of apoptosis and CaMKⅡ/mPTP pathway.
Journal of biochemical and molecular toxicology
Liu K,Hu C,Yin W,Zhou L,Gu X,Zuo X
Sun Jan 01 00:00:00 UTC 2023
Lipin2 ameliorates diabetic encephalopathy via suppressing JNK/ERK-mediated NLRP3 inflammasome overactivation.
International immunopharmacology
Zhang X,Huang S,Zhuang Z,Han X,Xie M,Yu S,Hua M,Liang Z,Meng C,Yin L,Zhuang X,Chen S
Mon May 01 00:00:00 UTC 2023
LINC00365 functions as a tumor suppressor by inhibiting HIF-1α-mediated glucose metabolism reprogramming in breast cancer.
Experimental cell research
Liu B,Qu X,Wang J,Xu L,Zhang L,Xu B,Su J,Bian X
Sat Apr 01 00:00:00 UTC 2023
miR-656-3p inhibits melanomas in vitro and in vivo by inducing senescence via inhibiting LMNB2.
Journal of cancer research and clinical oncology
Sun J,Lui K,Pang Q,Xu M,Zhao H,Shao J,Yu Y,Chu X,Liang Y,Xu J,Shen Z
Fri Sep 01 00:00:00 UTC 2023
DNA damage-inducible transcript 3 deficiency promotes bone resorption in murine periodontitis models.
Journal of periodontal research
Luo Y,Yang B,Dong W,Yu W,Jia M,Wang J
Tue Aug 01 00:00:00 UTC 2023
Dysfunction of inhibitory interneurons contributes to synaptic plasticity via GABABR-pNR2B signaling in a chronic migraine rat model.
Frontiers in molecular neuroscience
Zeng X,Niu Y,Qin G,Zhang D,Chen L
Mon Jun 19 00:00:00 UTC 2023
Combination treatment with FAAH inhibitors/URB597 and ferroptosis inducers significantly decreases the growth and metastasis of renal cell carcinoma cells via the PI3K-AKT signaling pathway.
Cell death & disease
Hao J,Chen Q,Feng Y,Jiang Q,Sun H,Deng B,Huang X,Guan J,Chen Q,Liu X,Wang Y,Cao P,Feng F,Li X
Thu Apr 06 00:00:00 UTC 2023
Autophagy ameliorates Pseudomonas aeruginosa-infected diabetic wounds by regulating the toll-like receptor 4/myeloid differentiation factor 88 pathway.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
Ji X,Jin P,Yu P,Wang P
Fri May 26 00:00:00 UTC 2023
Anti-inflammatory labdane diterpenoids from the aerial parts of Leonurus japonicus.
Phytochemistry
Wei QH,Cao XX,Xu DF,Wang ST,Zhang JS,Zhang H
Thu Jun 01 00:00:00 UTC 2023
Critical role of caveolin-1 in intestinal ischemia reperfusion by inhibiting protein kinase C βII.
Free radical biology & medicine
Chen Z,Wang Z,Liu D,Zhao X,Ning S,Liu X,Wang G,Zhang F,Luo F,Yao J,Tian X
Sun Jan 01 00:00:00 UTC 2023
Tartary buckwheat flavonoids alleviates high-fat diet induced kidney fibrosis in mice by inhibiting MAPK and TGF-β1/Smad signaling pathway.
Chemico-biological interactions
Liu S,Fu S,Jin Y,Geng R,Li Y,Zhang Y,Liu J,Guo W
Sat Jul 01 00:00:00 UTC 2023
Rapamycin inhibits corneal inflammatory response and neovascularization in a mouse model of corneal alkali burn.
Experimental eye research
Li J,Han J,Shi Y,Liu M
Tue Aug 01 00:00:00 UTC 2023
TGF-β Signaling Activation Confers Anlotinib Resistance in Gastric Cancer.
Pharmaceutical research
Chen J,Hai Y,Hu Q,Chen C,Jiang X,Gao Y
Wed Mar 01 00:00:00 UTC 2023
CMTM6 overexpression confers trastuzumab resistance in HER2-positive breast cancer.
Molecular cancer
Xing F,Gao H,Chen G,Sun L,Sun J,Qiao X,Xue J,Liu C
Tue Jan 10 00:00:00 UTC 2023
Silencing SOX11 Alleviates Allergic Rhinitis by Inhibiting Epithelial-Derived Cytokines.
Balkan medical journal
Jiang L,Wang C,Zhao R,Cao J,Liu Y,Tian L,Liu M
Mon Jan 23 00:00:00 UTC 2023
Inhibition of deubiquitinase USP28 attenuates cyst growth in autosomal dominant polycystic kidney disease.
Biochemical pharmacology
Ren Y,Zhu X,Fu K,Zhang H,Zhao W,Lin Y,Fang Q,Wang J,Chen Y,Guo D
Sun Jan 01 00:00:00 UTC 2023
Determination of the optimal concentration and duration of C5aR antagonist application in an inflammatory model of human dental pulp cells.
FEBS open bio
Hu J,Tan X,Wei X,Hu W,Gao L,Cao X,Yang H,Jiang Z,Li N,Teng L,Liu M
Wed Mar 01 00:00:00 UTC 2023
Systems pharmacology reveals the mechanism of Astragaloside IV in improving immune activity on cyclophosphamide-induced immunosuppressed mice.
Journal of ethnopharmacology
Yao J,Liu J,He Y,Liu L,Xu Z,Lin X,Liu N,Kai G
Fri Sep 15 00:00:00 UTC 2023
ERK Inhibition Promotes Engraftment of Allografts by Reprogramming T-Cell Metabolism.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Tan X,Qi C,Zhao X,Sun L,Wu M,Sun W,Gu L,Wang F,Feng H,Huang X,Xie B,Shi Z,Xie P,Wu M,Zhang Y,Chen G
Thu Jun 01 00:00:00 UTC 2023
2-Undecanone alleviates asthma by inhibiting NF-κB pathway.
Biochemistry and cell biology = Biochimie et biologie cellulaire
Song G,Yu S,Zhang Y,Sun M,Zhang B,Peng M
Wed Feb 01 00:00:00 UTC 2023
Novel ELAC2 Mutations in Individuals Presenting with Variably Severe Neurological Disease in the Presence or Absence of Cardiomyopathy.
Life (Basel, Switzerland)
Cafournet C,Zanin S,Guimier A,Hully M,Assouline Z,Barcia G,de Lonlay P,Steffann J,Munnich A,Bonnefont JP,Rötig A,Ruzzenente B,Metodiev MD
Sat Feb 04 00:00:00 UTC 2023
RNF123 Mediates Ubiquitination and Degradation of SOCS1 To Regulate Type I Interferon Production during Duck Tembusu Virus Infection.
Journal of virology
Huang S,Cui M,Huang J,Wu Z,Cheng A,Wang M,Zhu D,Chen S,Liu M,Zhao X,Wu Y,Yang Q,Zhang S,Ou X,Mao S,Gao Q,Tian B,Sun D,Yin Z,Jing B,Jia R
Thu Apr 27 00:00:00 UTC 2023
A new acid isolated from V. negundo L. inhibits NLRP3 inflammasome activation and protects against inflammatory diseases.
Frontiers in immunology
Di Q,Zhao X,Lin J,Li X,Li X,Tang H,Zhang R,Xiao W,Chen W
Tue May 09 00:00:00 UTC 2023
Effect of transmembrane protein 100 on prostate cancer progression by regulating SCNN1D through the FAK/PI3K/AKT pathway.
Translational oncology
Ye Z,Xia Y,Li L,Li B,Chen W,Han S,Zhou X,Chen L,Yu W,Ruan Y,Cheng F
Sun Jan 01 00:00:00 UTC 2023
Suprabasin enhances the invasion, migration, and angiogenic ability of oral squamous cell carcinoma cells under hypoxic conditions.
Oncology reports
Houri A,Mukudai Y,Abe Y,Watanabe M,Nara M,Miyamoto S,Kurihara M,Shimane T,Shirota T
Mon May 01 00:00:00 UTC 2023
The m6A Reader YTHDF1 Promotes Lung Carcinoma Progression via Regulating Ferritin Mediate Ferroptosis in an m6A-Dependent Manner.
Pharmaceuticals (Basel, Switzerland)
Diao H,Tan H,Hu Y,Wang R,Cai P,Huang B,Shao X,Yan M,Yin C,Zhang Y
Wed Jan 25 00:00:00 UTC 2023
Human umbilical cord mesenchymal stem cells conditioned medium exerts anti-tumor effects on KGN cells in a cell density-dependent manner through activation of the Hippo pathway.
Stem cell research & therapy
Wan W,Miao Y,Niu Y,Zhu K,Ma Y,Pan M,Ma B,Wei Q
Mon Mar 20 00:00:00 UTC 2023
SIRT3-mediated autophagy contributes to ferroptosis-induced anticancer by inducing the formation of BECN1-SLC7A11 complex.
Biochemical pharmacology
Huang P,Zhao H,Pan X,Li J,Pan W,Dai H,Wang J,Xia C,Liu F
Sat Jul 01 00:00:00 UTC 2023
Silica nanoparticles suppressed the spermatogenesis via downregulation of miR-450b-3p by targeting Layilin in spermatocyte of mouse.
Environmental pollution (Barking, Essex : 1987)
Zhou G,Wang J,Ren L,Liu J,Li X,Zhang Y,Sang Y,Gao L,Li Y,Sun Z,Zhou X
Wed Feb 01 00:00:00 UTC 2023
Swimming training combined with fecal microbial transplantation protects motor functions in rats with spinal cord injury by improving the intestinal system.
Neuroscience letters
Ouyang S,Wang X,Chen Y,Deng L,Yang X,Hu S,Wu S
Thu Mar 16 00:00:00 UTC 2023
Analysis of Promoter Methylation of the Bovine FOXO1 Gene and Its Effect on Proliferation and Differentiation of Myoblasts.
Animals : an open access journal from MDPI
Shi P,Ruan Y,Liu W,Sun J,Xu J,Xu H
Mon Jan 16 00:00:00 UTC 2023
Deciphering the molecular mechanism of the THBS1 gene in the TNF signaling axis in glioma stem cells.
Cellular signalling
Chen L,Fang W,Chen W,Wei Y,Ding J,Li J,Lin J,Wu Q
Thu Jun 01 00:00:00 UTC 2023
Research Note: Duck plague virus pUL48 is a late protein that plays an important role in viral replication.
Poultry science
Zhou T,Wang M,Ruan P,Fan D,Cheng A,Zhang W,Tian B,Yang Q,Wu Y,Zhang S,Ou X,Mao S,Huang J,Gao Q,Sun D,Zhao X,Chen S,Liu M,Zhu D,Jia R
Wed Feb 01 00:00:00 UTC 2023
JNK-JUN-NCOA4 axis contributes to chondrocyte ferroptosis and aggravates osteoarthritis via ferritinophagy.
Free radical biology & medicine
Sun K,Hou L,Guo Z,Wang G,Guo J,Xu J,Zhang X,Guo F
Mon May 01 00:00:00 UTC 2023
Multiomics analyses and machine learning of nuclear receptor coactivator 6 reveal its essential role in hepatocellular carcinoma.
Cancer science
Fang Y,Xu Y,Liao W,Ji T,Yu L,Li L,Pan M,Yang D
Sun Jan 01 00:00:00 UTC 2023
Neutrophil membrane-camouflaged nanoparticles alleviate inflammation and promote angiogenesis in ischemic myocardial injury.
Bioactive materials
Han D,Wang F,Qiao Z,Wang B,Zhang Y,Jiang Q,Liu M,Zhuang Y,An Q,Bai Y,Shangguan J,Zhang J,Liang G,Shen D
Mon May 01 00:00:00 UTC 2023
SERPINF1 Mediates Tumor Progression and Stemness in Glioma.
Genes
Song L,Huo X,Li X,Xu X,Zheng Y,Li D,Zhang J,Wang K,Wang L,Wu Z
Sat Feb 25 00:00:00 UTC 2023
Proteasomal inhibitors induce myeloma cell pyroptosis via the BAX/GSDME pathway.
Acta pharmacologica Sinica
Liang JP,He YM,Cui YL,Sun YN,He GS,Zhu ZG,Mao XL
Sat Jul 01 00:00:00 UTC 2023
CENPH overexpression promotes the progression, cisplatin resistance, and poor prognosis of lung adenocarcinoma via the AKT and ERK/P38 pathways.
American journal of cancer research
Li Q,Huang X,Li QY,Tang Y,Liang LB,Luo Q,Gong M,Song YX,Guo Q,Chen C
Mon Jun 12 00:00:00 UTC 2023
Porous composite hydrogels with improved MSC survival for robust epithelial sealing around implants and M2 macrophage polarization.
Acta biomaterialia
Li Y,Zhang J,Wang C,Jiang Z,Lai K,Wang Y,Yang G
Wed Feb 01 00:00:00 UTC 2023
Ketogenic diet alleviates renal interstitial fibrosis in UUO mice by regulating macrophage proliferation.
The Journal of nutritional biochemistry
Hu X,Qiu Y,Cao R,Xu C,Lu C,Wang Z,Yang J
Tue Aug 01 00:00:00 UTC 2023
MED27 plays a tumor-promoting role in breast cancer progression by targeting KLF4.
Cancer science
Wang R,Yu W,Zhu T,Lin F,Hua C,Ru L,Guo P,Wan X,Xue G,Guo Z,Han S,Lv K,Zhang G,Ge H,Guo W,Xu L,Deng W
Thu Jun 01 00:00:00 UTC 2023
Resveratrol suppresses neuroinflammation to alleviate mechanical allodynia by inhibiting Janus kinase 2/signal transducer and activator of transcription 3 signaling pathway in a rat model of spinal cord injury.
Frontiers in molecular neuroscience
Han J,Hua Z,Yang WJ,Wang S,Yan F,Wang JN,Sun T
Tue Mar 07 00:00:00 UTC 2023
Sp1 promotes tumour progression by remodelling the mitochondrial network in cervical cancer.
Journal of translational medicine
Xu X,Wang X,Chen Q,Zheng A,Li D,Meng Z,Li X,Cai H,Li W,Huang S,Wang F
Sat May 06 00:00:00 UTC 2023
Construction and Characterization of Immortalized Fibroblast Cell Line from Bactrian Camel.
Life (Basel, Switzerland)
Yan M,Yong F,Ji W,Zhang L,Zhao S,Gao Y
Wed Jun 07 00:00:00 UTC 2023
M2 Macrophage Membrane-Mediated Biomimetic-Nanoparticle Carrying COX-siRNA Targeted Delivery for Prevention of Tendon Adhesions by Inhibiting Inflammation.
Small (Weinheim an der Bergstrasse, Germany)
Sun J,Ju F,Jin J,Wang HL,Li ZJ,Sun YC,Chen QZ,Yang QQ,Tan J,Zhou YL
Tue Aug 01 00:00:00 UTC 2023
Near-Infrared Phototheranostic Iron Pyrite Nanocrystals Simultaneously Induce Dual Cell Death Pathways via Enhanced Fenton Reactions in Triple-Negative Breast Cancer.
ACS nano
Zhao C,Liu Z,Chang CC,Chen YC,Zhang Q,Zhang XD,Andreou C,Pang J,Liu ZX,Wang DY,Kircher MF,Yang J
Tue Mar 14 00:00:00 UTC 2023
The role of CNPY2 in endothelial injury and inflammation during the progress of atherosclerosis.
Journal of molecular histology
Huang H,Tang N,Li Y,Huo Q,Chen Q,Meng Q
Thu Jun 01 00:00:00 UTC 2023
ML390 inhibits enterovirus 71 replication by targeting de novo pyrimidine biosynthesis pathway.
Antiviral research
Yang Q,Wu C,Zhu G,Ren F,Lin B,Huang R,Hu X,Zhao D,Peng K,Wu Y,Wang Q,Huang C,Zhang D
Sun Jan 01 00:00:00 UTC 2023
Lactoferrin Relieves Deoxynivalenol-Induced Oxidative Stress and Inflammatory Response by Modulating the Nrf2/MAPK Pathways in the Liver.
Journal of agricultural and food chemistry
Hu P,Liu Y,Li S,Zhao Y,Gu H,Zong Q,Ahmed AA,Bao W,Liu HY,Cai D
Wed May 31 00:00:00 UTC 2023
Bis-Benzylisoquinoline Alkaloids Inhibit Porcine Epidemic Diarrhea Virus by Disrupting Virus Entry.
Pathogens (Basel, Switzerland)
Zhang C,Chen H,Sun L,Zhao P,Qi C,Yang Y,Si A,Qian Y,Jung YS
Mon Jun 19 00:00:00 UTC 2023
MiR-124 Reduced Neuroinflammation after Traumatic Brain Injury by Inhibiting TRAF6.
Neuroimmunomodulation
Yang Y,Ye Y,Fan K,Luo J,Yang Y,Ma Y
Tue Dec 26 00:00:00 UTC 2023
PCSK9 Promotes Endothelial Dysfunction During Sepsis Via the TLR4/MyD88/NF-κB and NLRP3 Pathways.
Inflammation
Huang L,Li Y,Cheng Z,Lv Z,Luo S,Xia Y
Wed Feb 01 00:00:00 UTC 2023
Effects of chronic sleep restriction on the neuro-phenotypes of Ctnnd2 knockout mice.
Brain and behavior
Xu M,Wang X,Wang L,Wang S,Deng J,Wang Y,Li Y,Pan S,Liao A,Tao Y,Tan S
Sat Jul 01 00:00:00 UTC 2023
N-acetyl-L-cysteine alleviates FUNDC1-mediated mitophagy by regulating mitochondrial dynamics in type 1 diabetic nephropathy canine.
Life sciences
Ma F,Li H,Huo H,Han Q,Liao J,Zhang H,Li Y,Pan J,Hu L,Guo J,Tang Z
Sun Jan 15 00:00:00 UTC 2023
Hydrogen sulfide alleviates beryllium sulfate-induced ferroptosis and ferritinophagy in 16HBE cells.
Journal of applied toxicology : JAT
Liu X,Li G,Chen S,Jin H,Liu X,Zhang L,Zhang Z
Tue Aug 01 00:00:00 UTC 2023
Platelet-derived extracellular vesicles ameliorate intervertebral disc degeneration by alleviating mitochondrial dysfunction.
Materials today. Bio
Dai Z,Xia C,Zhao T,Wang H,Tian H,Xu O,Zhu X,Zhang J,Chen P
Wed Feb 01 00:00:00 UTC 2023
Exogenous Hydrogen Sulfide Induces A375 Melanoma Cell Apoptosis Through Overactivation of the Unfolded Protein Response.
Clinical, cosmetic and investigational dermatology
Ma W,Zhang X,Zhuang L
Tue Jul 04 00:00:00 UTC 2023
Significance of UGT1A6, UGT1A9, and UGT2B7 genetic variants and their mRNA expression in the clinical outcome of renal cell carcinoma.
Molecular and cellular biochemistry
Matsumoto J,Nishimoto A,Watari S,Ueki H,Shiromizu S,Iwata N,Takeda T,Ushio S,Kajizono M,Fujiyoshi M,Koyama T,Araki M,Wada K,Zamami Y,Nasu Y,Ariyoshi N
Tue Aug 01 00:00:00 UTC 2023
RRP Regulates Autophagy through the AMPK Pathway to Alleviate the Effect of Cell Senescence on Atherosclerosis.
Oxidative medicine and cellular longevity
Liu D,Song Y,Song T,Lin L,Zhang L,Yang Q,Niu X,Liang D,Yin J,Yang W,Zhang D
Fri Feb 10 00:00:00 UTC 2023
Lipophagy-mediated cholesterol synthesis inhibition is required for the survival of hepatocellular carcinoma under glutamine deprivation.
Redox biology
Kong Y,Wu M,Wan X,Sun M,Zhang Y,Wu Z,Li C,Liang X,Gao L,Ma C,Yue X
Sat Jul 01 00:00:00 UTC 2023
Sea Cucumber Hydrolysate Alleviates Immunosuppression and Gut Microbiota Imbalance Induced by Cyclophosphamide in Balb/c Mice through the NF-κB Pathway.
Foods (Basel, Switzerland)
Mao J,Li S,Fu R,Wang Y,Meng J,Jin Y,Wu T,Zhang M
Mon Apr 10 00:00:00 UTC 2023
Profilin1 Promotes Renal Tubular Epithelial Cell Apoptosis in Diabetic Nephropathy Through the Hedgehog Signaling Pathway.
Diabetes, metabolic syndrome and obesity : targets and therapy
Mai L,He G,Chen J,Zhu J,Chen S,Yang H,Zhang M,Hou X,Ke M,Li X
Mon Jun 19 00:00:00 UTC 2023
TRPM2 facilitates tumor progression of clear cell renal cell carcinoma by relieving Endoplasmic Reticulum Stress.
International journal of medical sciences
Yuan H,Li W,Lou N,Yu T,Meng X,Xiao W,Zhang X
Tue Jan 10 00:00:00 UTC 2023
Enhanced autophagy reversed aflatoxin B1-induced decrease in lactate secretion of dairy goat Sertoli cells.
Ecotoxicology and environmental safety
Han S,Zhang H,Liu X,Wen F,Li B,Bie Z,Qiu H,Hu J
Sat Jul 01 00:00:00 UTC 2023
KRT17 Promotes T-lymphocyte Infiltration Through the YTHDF2-CXCL10 Axis in Colorectal Cancer.
Cancer immunology research
Liang W,Liu H,Zeng Z,Liang Z,Xie H,Li W,Xiong L,Liu Z,Chen M,Jie H,Zheng X,Huang L,Kang L
Wed Jul 05 00:00:00 UTC 2023
Multimodal rehabilitation promotes axonal sprouting and functional recovery in a murine model of spinal cord injury (SCI).
Neuroscience letters
Wang Y,Luo H,Liu Y,Yang C,Yin Y,Tan B
Tue Jan 31 00:00:00 UTC 2023
Inhibition of cGAS aggravated the host inflammatory response to Aspergillus fumigatus.
Experimental lung research
Peng M,Li X,Zhang X,Peng L
Sat Dec 16 00:00:00 UTC 2023
Single-cell sequencing reveals the immune microenvironment landscape related to anti-PD-1 resistance in metastatic colorectal cancer with high microsatellite instability.
BMC medicine
Wu T,Zhang X,Liu X,Cai X,Shen T,Pan D,Liang R,Ding R,Hu R,Dong J,Li F,Li J,Xie L,Wang C,Geng S,Yang Z,Xing L,Li Y
Thu Apr 27 00:00:00 UTC 2023
A novel Hoxd13 mutation causes synpolydactyly and promotes osteoclast differentiation by regulating pSmad5/p65/c-Fos/Rank axis.
Cell death & disease
Zhang L,Fang Z,Cheng G,He M,Lin Y
Mon Feb 20 00:00:00 UTC 2023
CENPF promotes the proliferation of renal cell carcinoma in vitro.
Translational andrology and urology
Zhang J,Wang Z,Liu Z,Chen Z,Jiang J,Ji Y,Zhang Y,Zhu H,Zheng B
Tue Feb 28 00:00:00 UTC 2023
Myocardial ischemia-reperfusion injury is probably due to the excessive production of mitochondrial ROS caused by the activation of 5-HT degradation system mediated by PAF receptor.
Molecular immunology
Jin J,Xu F,Zhang Y,Guan J,Fu J
Wed Mar 01 00:00:00 UTC 2023
Exploring the effect of polyamines on NK cell function in colorectal cancer process based on glycolysis.
International immunopharmacology
He H,Song Z,Lin S,Wang Y,Wang G
Sat Apr 01 00:00:00 UTC 2023
NLRP3 activation in microglia contributes to learning and memory impairment induced by chronic lead exposure in mice.
Toxicological sciences : an official journal of the Society of Toxicology
Zhu J,Zhou F,Zhou Q,Xu Y,Li Y,Huang D,Chen L,Liu A,Zou F,Meng X
Tue Jan 31 00:00:00 UTC 2023
Insight into mitochondrial dysfunction mediated by clozapine-induced inhibition of PGRMC1 in PC12 cells.
Toxicology
Wang L,Chen Q,Ma R,Zhang B,Yang P,Cao T,Jiao S,Chen H,Lin C,Cai H
Thu Jun 01 00:00:00 UTC 2023
Photodynamic therapy enhances the cytotoxicity of temozolomide against glioblastoma via reprogramming anaerobic glycolysis.
Photodiagnosis and photodynamic therapy
Li Y,Wang D,Zhang Z,Wang Y,Zhang Z,Liang Z,Liu F,Chen L
Thu Jun 01 00:00:00 UTC 2023
Discovery of potent and selective PI3Kδ inhibitors bearing amino acid fragments.
Bioorganic chemistry
Lei H,Duan W,Zhang SQ,Feng Y,Ma M,Yuan B,Xin M
Fri Sep 01 00:00:00 UTC 2023
YAP and β-catenin cooperate to drive H. pylori-induced gastric tumorigenesis.
Gut microbes
Li N,Xu X,Zhan Y,Fei X,Ouyang Y,Zheng P,Zhou Y,He C,Xie C,Hu Y,Hong J,Lu N,Ge Z,Zhu Y
Mon Mar 27 00:00:00 UTC 2023
miR-342-5p downstream to Notch enhances arterialization of endothelial cells in response to shear stress by repressing MYC.
Molecular therapy. Nucleic acids
Zhang X,Sun J,Zhang P,Wen T,Wang R,Liang L,Yang Z,Li J,Zhang J,Che B,Feng X,Liu X,Han H,Yan X
Tue Jun 13 00:00:00 UTC 2023
Caveolae-associated protein 3 promotes adipogenic differentiation of porcine preadipocytes by promoting extracellular signal-regulated kinase phosphorylation.
Animal science journal = Nihon chikusan Gakkaiho
Shi M,Yang S,Huang X,Wang S,Li W,Yun J,Lu C,Yang Y,Cai C,Gao P,Guo X,Li B,Cao G
Mon Mar 20 00:00:00 UTC 2023
Knock-down of PGM1 inhibits cell viability, glycolysis, and oxidative phosphorylation in glioma under low glucose condition via the Myc signaling pathway.
Biochemical and biophysical research communications
Liu S,Deng Y,Yu Y,Xia X
Sun May 14 00:00:00 UTC 2023
Persistent Müllerian duct syndrome in an assisted reproductive patient: a novel variant impairs the biosynthesis and secretion of anti-Müllerian hormone (AMH).
Asian journal of andrology
Du Q,Qiu C,Zhang L,Wang QY,Hong K,Liu XL
Fri Jul 21 00:00:00 UTC 2023
Ccdc25 regulates neurogenesis during the brain development.
Developmental neurobiology
Wang C,Qin J,Jiao J,Ji F
Mon May 22 00:00:00 UTC 2023
Elevated FBXL18 promotes RPS15A ubiquitination and SMAD3 activation to drive HCC.
Hepatology communications
Yu HQ,Li F,Xiong H,Fang L,Zhang J,Bie P,Xie CM
Sat Jul 01 00:00:00 UTC 2023
Identification of novel candidate targets for suppressing ovarian cancer progression through IL-33/ST2 axis components using the system biology approach.
Frontiers in molecular biosciences
Reivan Ortiz GG,Ciongradi CI,Chaitanya MVNL,Narayanan J,Mohany M,Al-Rejaie SS,Arias-Gonzáles JL,Sârbu I,Assefi M,Akram SV,Döğüş Y,Bahrami A,Akhavan-Sigari R
Tue Jun 20 00:00:00 UTC 2023
Intraoperative blood auto-transfusion restrained the malignancy of Liver Cancer via regulating functions of tumor cells and Kuffer cells.
Biotechnology & genetic engineering reviews
Yao N,Wu M,You L,Xu J,Liu T,Wang J,Li Z,Guo J,Sun Y
Thu May 18 00:00:00 UTC 2023
Wei-Tong-Xin ameliorated cisplatin-induced mitophagy and apoptosis in gastric antral mucosa by activating the Nrf2/HO-1 pathway.
Journal of ethnopharmacology
Zhang X,Wang S,Jin Y,Wang J,Wang R,Yang X,Zhang S,Yan T,Jia Y
Tue May 23 00:00:00 UTC 2023
Pseurotin A Validation as a Metastatic Castration-Resistant Prostate Cancer Recurrence-Suppressing Lead via PCSK9-LDLR Axis Modulation.
Marine drugs
Abdelwahed KS,Siddique AB,Ebrahim HY,Qusa MH,Mudhish EA,Rad AH,Zerfaoui M,Abd Elmageed ZY,El Sayed KA
Tue Mar 28 00:00:00 UTC 2023
BDE209-promoted Dio2 degradation in H4 glioma cells through the autophagy pathway, resulting in hypothyroidism and leading to neurotoxicity.
Toxicology
Liu M,Yu Z,Yang F,Zhao Z,Zhou M,Wang C,Zhang B,Liang G,Liu X,Shao J
Tue Aug 01 00:00:00 UTC 2023
Regulation of telomere silencing by the core histones-autophagy-Sir2 axis.
Life science alliance
Mei Q,Yu Q,Li X,Chen J,Yu X
Wed Mar 01 00:00:00 UTC 2023
ATF5 regulates tubulointerstitial injury in diabetic kidney disease via mitochondrial unfolded protein response.
Molecular medicine (Cambridge, Mass.)
Liu Y,Zhang L,Zhang S,Liu J,Li X,Yang K,Yang D,Liu Y,Sun L,Liu F,Xiao L
Mon Apr 24 00:00:00 UTC 2023
SOX9/NFIA promotes human ovarian cancer metastasis through the Wnt/β-catenin signaling pathway.
Pathology, research and practice
Lu R,Tang P,Zhang D,Lin S,Li H,Feng X,Sun M,Zhang H
Tue Aug 01 00:00:00 UTC 2023
ROS-activated MAPK/ERK pathway regulates crosstalk between Nrf2 and Hif-1α to promote IL-17D expression protecting the intestinal epithelial barrier under hyperoxia.
International immunopharmacology
Wang P,Li T,Niu C,Sun S,Liu D
Wed Mar 01 00:00:00 UTC 2023
Integrated analysis of cell-specific gene expression in peripheral blood using ISG15 as a marker of rejection in kidney transplantation.
Frontiers in immunology
Zhang Z,Qin Y,Wang Y,Li S,Hu X
Fri Mar 31 00:00:00 UTC 2023
A Water-Soluble Hydrogen Sulfide Donor Suppresses the Growth of Hepatocellular Carcinoma via Inhibiting the AKT/GSK-3β/β-Catenin and TGF-β/Smad2/3 Signaling Pathways.
Journal of oncology
Duan SF,Zhang MM,Dong Q,Yang B,Liu W,Zhang X,Yu HL,Zhang SH,Khan NH,Wu DD,Zhang XJ,Cen J
Sat Mar 18 00:00:00 UTC 2023
ITGB1-mediated molecular landscape and cuproptosis phenotype induced the worse prognosis in diffuse gastric cancer.
Frontiers in oncology
Zhu X,Chen H,Li H,Ren H,Ye C,Xu K,Liu J,Du F,Zhang Z,Liu Y,Xie X,Wang M,Ma T,Chong W,Shang L,Li L
Tue Apr 04 00:00:00 UTC 2023
Plasma microRNA expression profiles associated with zinc exposure and type 2 diabetes mellitus: Exploring potential role of miR-144-3p in zinc-induced insulin resistance.
Environment international
Ye Z,Cheng M,Fan L,Ma J,Zhang Y,Gu P,Xie Y,You X,Zhou M,Wang B,Chen W
Wed Feb 01 00:00:00 UTC 2023
Neuroprotective Effects of Sodium Butyrate by Restoring Gut Microbiota and Inhibiting TLR4 Signaling in Mice with MPTP-Induced Parkinson's Disease.
Nutrients
Guo TT,Zhang Z,Sun Y,Zhu RY,Wang FX,Ma LJ,Jiang L,Liu HD
Mon Feb 13 00:00:00 UTC 2023
UBE2E2 enhances Snail-mediated epithelial-mesenchymal transition and Nrf2-mediated antioxidant activity in ovarian cancer.
Cell death & disease
Hong X,Ma N,Li D,Zhang M,Dong W,Huang J,Ci X,Zhang S
Fri Feb 10 00:00:00 UTC 2023
Knockdown of CENPN Inhibits Glucose Metabolism and Induces G1 Arrest in Esophageal Cancer Cells by Regulating PI3K/AKT Signaling Pathway.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme
Xu S,Wei X
Tue Aug 01 00:00:00 UTC 2023
Glycolysis-related lncRNA TMEM105 upregulates LDHA to facilitate breast cancer liver metastasis via sponging miR-1208.
Cell death & disease
Han J,Chen X,Wang J,Liu B
Fri Feb 03 00:00:00 UTC 2023
SLC27A2 mediates FAO in colorectal cancer through nongenic crosstalk regulation of the PPARs pathway.
BMC cancer
Shang K,Ma N,Che J,Li H,Hu J,Sun H,Cao B
Tue Apr 11 00:00:00 UTC 2023
Neuroprotection of NSC Therapy is Superior to Glibenclamide in Cardiac Arrest-Induced Brain Injury via Neuroinflammation Regulation.
Translational stroke research
Wang Z,Zhang S,Du J,Lachance BB,Chen S,Polster BM,Jia X
Sun Oct 01 00:00:00 UTC 2023
Methyl gallate prevents oxidative stress induced apoptosis and ECM degradation in chondrocytes via restoring Sirt3 mediated autophagy and ameliorates osteoarthritis progression.
International immunopharmacology
Li Y,Shen B,Lv C,Zhu X,Naren Q,Xu D,Chen H,Wu F
Sun Jan 01 00:00:00 UTC 2023
USP33 enhances cell survival and stemness by deubiquitinating CTNNB1 in BXPC-3 and SW1990 cells.
Cell biology international
Zhong B,Zhu Q,Wang L,Wang F,Zheng Y,Lin S,Cheng X
Tue Aug 01 00:00:00 UTC 2023
Porcine reproductive and respiratory syndrome virus infection triggers autophagy via ER stress-induced calcium signaling to facilitate virus replication.
PLoS pathogens
Diao F,Jiang C,Sun Y,Gao Y,Bai J,Nauwynck H,Wang X,Yang Y,Jiang P,Liu X
Wed Mar 01 00:00:00 UTC 2023
The nanoprodrug of polytemozolomide combines with MGMT siRNA to enhance the effect of temozolomide in glioma.
Drug delivery
Xu H,Zhang Y,Li L,Ren Y,Qian F,Wang L,Ma H,Quan A,Liu H,Yu R
Fri Dec 01 00:00:00 UTC 2023
18β-Glycyrrhetinic Acid Alleviates P. multocida-Induced Vascular Endothelial Inflammation by PARP1-Mediated NF-κB and HMGB1 Signalling Suppression in PIEC Cells.
Infection and drug resistance
Lu Q,Han W,Wen D,Guo P,Liu Y,Wu Z,Fu S,Ye C,Wang X,Qiu Y
Tue Jul 18 00:00:00 UTC 2023
Huc-MSCs-derived exosomes attenuate neuropathic pain by inhibiting activation of the TLR2/MyD88/NF-κB signaling pathway in the spinal microglia by targeting Rsad2.
International immunopharmacology
Gao X,Gao LF,Zhang YN,Kong XQ,Jia S,Meng CY
Sun Jan 01 00:00:00 UTC 2023
Cryptotanshinone modulates proliferation, apoptosis, and fibrosis through inhibiting AR and EGFR/STAT3 axis to ameliorate benign prostatic hyperplasia progression.
European journal of pharmacology
Wei P,Lin D,Zhang M,Luo C,Wu X,Deng B,Cui K,Chen Z
Thu Jan 05 00:00:00 UTC 2023
Stevioside targets the NF-κB and MAPK pathways for inhibiting inflammation and apoptosis of chondrocytes and ameliorates osteoarthritis in vivo.
International immunopharmacology
Cai T,Ye H,Jiang H,Lin C,Lou C,Wang W,Yan Z,Xue X,Pan X,Lin J
Wed Feb 01 00:00:00 UTC 2023
Proximal tubular Bmal1 protects against chronic kidney injury and renal fibrosis by maintaining of cellular metabolic homeostasis.
Biochimica et biophysica acta. Molecular basis of disease
Liu C,Li S,Ji S,Zhang J,Zheng F,Guan Y,Yang G,Chen L
Sun Jan 01 00:00:00 UTC 2023
Oleoylethanolamide alleviates hyperlipidaemia-mediated vascular calcification via attenuating mitochondrial DNA stress triggered autophagy-dependent ferroptosis by activating PPARα.
Biochemical pharmacology
Chen Z,Sun X,Li X,Liu N
Wed Feb 01 00:00:00 UTC 2023
Ferroptosis is involved in PM2.5-induced acute nasal epithelial injury via AMPK-mediated autophagy.
International immunopharmacology
Gu W,Hou T,Zhou H,Zhu L,Zhu W,Wang Y
Wed Feb 01 00:00:00 UTC 2023
Mutation-Driven S100A8 Overexpression Confers Aberrant Phenotypes in Type 1 CALR-Mutated MPN.
International journal of molecular sciences
Wang YH,Chen YJ,Lai YH,Wang MC,Chen YY,Wu YY,Yang YR,Tsou HY,Li CP,Hsu CC,Huang CE,Chen CC
Sun May 14 00:00:00 UTC 2023
Zika virus replication on endothelial cells and invasion into the central nervous system by inhibiting interferon β translation.
Virology
Wang K,Zou S,Chen H,Higazy D,Gao X,Zhang Y,Cao S,Cui M
Mon May 01 00:00:00 UTC 2023
RNA N6-methyladenosine reader IGF2BP3 interacts with MYCN and facilitates neuroblastoma cell proliferation.
Cell death discovery
Zhu K,Gao T,Wang Z,Zhang L,Tan K,Lv Z
Mon May 08 00:00:00 UTC 2023
The PRMT5 inhibitor C9 mitigates hypoxia-induced carboplatin resistance in lung cancer by inducing autophagy.
Cell biology international
Fan J,Li H,Ruan Q,Zhu X,Jing P,Gu Z
Sun Oct 01 00:00:00 UTC 2023
Benzothiazole Substitution Analogs of Rhodacyanine Hsp70 Inhibitors Modulate Tau Accumulation.
ACS chemical biology
Hill SE,Beaulieu-Abdelahad D,Lemus A,Webster JM,Ospina SR,Darling AL,Martin MD,Patel S,Bridenstine L,Swonger R,Paul S,Blackburn R,Calcul L,Dickey CA,Leahy JW,Blair LJ
Fri May 19 00:00:00 UTC 2023
Progressive degeneration of the retina in Loxl3 mutant mouse model of Stickler syndrome.
Developmental biology
Liu Z,Mo F,Dong X,Chen G,Gao J,Zhang J
Wed Mar 01 00:00:00 UTC 2023
Dendrobine Alleviates Cellular Senescence and Osteoarthritis via the ROS/NF-κB Axis.
International journal of molecular sciences
Chen H,Tu M,Liu S,Wen Y,Chen L
Wed Jan 25 00:00:00 UTC 2023
Garlic-derived exosomes regulate PFKFB3 expression to relieve liver dysfunction in high-fat diet-fed mice via macrophage-hepatocyte crosstalk.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu J,Li W,Bian Y,Jiang X,Zhu F,Yin F,Yin L,Song X,Guo H,Liu J
Sat Apr 01 00:00:00 UTC 2023
Chromatin remodeling and nucleoskeleton synergistically control osteogenic differentiation in different matrix stiffnesses.
Materials today. Bio
Xu X,Zhang H,Li Y,Liu F,Jing Z,Ren M,Chen T,Fu Y,Wu Y,Ji P,Yang S
Thu Jun 01 00:00:00 UTC 2023
Tandem mass tag-based quantitative proteomics analysis reveals the effects of the α-lactalbumin peptides GINY and DQW on lipid deposition and oxidative stress in HepG2 cells.
Journal of dairy science
Chen H,Qi X,Guan K,Wang R,Li Q,Ma Y
Sat Apr 01 00:00:00 UTC 2023
ARHGAP44 expression is associated with the metastasis of osteosarcoma and is a promising prognostic biomarker.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Hu S,Wang W
Thu Jun 01 00:00:00 UTC 2023
Tumor-derived endomucin promotes colorectal cancer proliferation and metastasis.
Cancer medicine
Huang Q,Li XM,Sun JP,Zhou Y
Wed Feb 01 00:00:00 UTC 2023
Empagliflozin-Pretreated Mesenchymal Stem Cell-Derived Small Extracellular Vesicles Attenuated Heart Injury.
Oxidative medicine and cellular longevity
Chi B,Zou A,Mao L,Cai D,Xiao T,Wang Y,Wang Q,Ji Y,Sun L
Wed Mar 01 00:00:00 UTC 2023
Mulberry leaves attenuate D-galactose-induced aging in vivo and in vitro.
Journal of ethnopharmacology
Zhu Y,Han Y,Wang W,Liang G,Qi J
Sat Jul 15 00:00:00 UTC 2023
Blockade of neutrophil extracellular traps ameliorates toluene diisocyanate-induced steroid-resistant asthma.
International immunopharmacology
Peng X,Li Y,Zhao W,Yang S,Huang J,Chen Y,Wang Y,Gong Z,Chen X,Yu C,Cai S,Zhao H
Sat Apr 01 00:00:00 UTC 2023
Ubiquitin ligase MARCH8 promotes the malignant progression of hepatocellular carcinoma through PTEN ubiquitination and degradation.
Molecular carcinogenesis
Xu Y,Zhang D,Ji J,Zhang L
Sat Jul 01 00:00:00 UTC 2023
Vitamin D-VDR (vitamin D receptor) alleviates glucose metabolism reprogramming in lipopolysaccharide-induced acute kidney injury.
Frontiers in physiology
Dai Q,Zhang H,Tang S,Wu X,Wang J,Yi B,Liu J,Li Z,Liao Q,Li A,Liu Y,Zhang W
Tue Mar 14 00:00:00 UTC 2023
The well-developed actin cytoskeleton and Cthrc1 expression by actin-binding protein drebrin in myofibroblasts promote cardiac and hepatic fibrosis.
The Journal of biological chemistry
Hironaka T,Takizawa N,Yamauchi Y,Horii Y,Nakaya M
Wed Mar 01 00:00:00 UTC 2023
Impact of age on the rotenone-induced sporadic Parkinson's disease model using Drosophila melanogaster.
Neuroscience letters
Li W,Pan X,Li M,Ling L,Zhang M,Liu Z,Zhang K,Guo J,Wang H
Sun May 14 00:00:00 UTC 2023
Toosendanin-induced apoptosis of CMT-U27 is mediated through the mitochondrial apoptotic pathway.
Veterinary and comparative oncology
Yang Y,Mei C,Xian H,Zhang X,Li J,Liang ZX,Zhi Y,Ma Y,Wang HJ
Thu Jun 01 00:00:00 UTC 2023
Synergistic anticancer effect of flavonoids from Sophora alopecuroides with Sorafenib against hepatocellular carcinoma.
Phytotherapy research : PTR
Zhu XF,Sun ZL,Ma J,Hu B,Yu MC,Liu XJ,Yang P,Xu Y,Ju D,Mu Q
Wed Feb 01 00:00:00 UTC 2023
Inhibition of mGluR5 ameliorates lipid accumulation and inflammation in HepG2 cells.
Biochemical and biophysical research communications
Zeng S,Tao M,Yuan L,Zhang L,Luo X
Sun Apr 23 00:00:00 UTC 2023
Dach1 deficiency drives alveolar epithelium apoptosis in pulmonary fibrosis via modulating C-Jun/Bim activity.
Translational research : the journal of laboratory and clinical medicine
Lu Y,Tang K,Wang S,Tian Z,Fan Y,Li B,Wang M,Zhao J,Xie J
Sat Jul 01 00:00:00 UTC 2023
Metabolomic and transcriptomic studies of improvements in myocardial infarction due to Pycr1 deletion.
Journal of cellular and molecular medicine
Xue Z,Pan Y,Kong X,Zhang J,Wu D,Zhou B
Sun Jan 01 00:00:00 UTC 2023
Lacosamide alleviates bilateral cavernous nerve injury-induced erectile dysfunction in the rat model by ameliorating pathological changes in the corpus cavernosum.
International journal of impotence research
Yao L,Cong R,Zhou X,Zhou X,Ji C,Wei X,Meng X,Song N
Wed Mar 15 00:00:00 UTC 2023
Hydrogen sulfide attenuates TMAO‑induced macrophage inflammation through increased SIRT1 sulfhydration.
Molecular medicine reports
Liu MH,Lin XL,Xiao LL
Sat Jul 01 00:00:00 UTC 2023
Decreasing REDD1 expression protects against high glucose-induced apoptosis, oxidative stress and inflammatory injury in podocytes through regulation of the AKT/GSK-3β/Nrf2 pathway.
Immunopharmacology and immunotoxicology
Wang X,Yang J,Wang W,Li Y,Yang Y
Sun Oct 01 00:00:00 UTC 2023
The total polyphenolic glycoside extract of Lamiophlomis rotata ameliorates hepatic fibrosis through apoptosis by TGF-β/Smad signaling pathway.
Chinese medicine
Wan G,Chen Z,Lei L,Geng X,Zhang Y,Yang C,Cao W,Pan Z
Fri Feb 24 00:00:00 UTC 2023
Phosphoglycerate dehydrogenase activates PKM2 to phosphorylate histone H3T11 and attenuate cellular senescence.
Nature communications
Wu Y,Tang L,Huang H,Yu Q,Hu B,Wang G,Ge F,Yin T,Li S,Yu X
Fri Mar 10 00:00:00 UTC 2023
Role of Protein Tyrosine Phosphatase 1B Inhibitor in Early Brain Injury of Subarachnoid Hemorrhage in Mice.
Brain sciences
Zhang ZH,Zhou XM,Zhang X
Thu May 18 00:00:00 UTC 2023
The involvement of let-7 in hCG-induced progesterone synthesis via regulating p27Kip1 and p21Cip1 expression.
Molecular and cellular endocrinology
Chen J,Liu K,Liu W,Yeung WS
Tue Aug 01 00:00:00 UTC 2023
NADPH-dependent ROS accumulation contributes to the impaired osteogenic differentiation of periodontal ligament stem cells under high glucose conditions.
Frontiers in endocrinology
Zhang YL,An Y,Sun LJ,Qu HL,Li X,He XT,Wu RX,Chen FM,Tian BM,Yin Y
Mon Jun 26 00:00:00 UTC 2023
IFN-γ Facilitates Corneal Epithelial Cell Pyroptosis Through the JAK2/STAT1 Pathway in Dry Eye.
Investigative ophthalmology & visual science
Yang X,Zuo X,Zeng H,Liao K,He D,Wang B,Yuan J
Wed Mar 01 00:00:00 UTC 2023
Combination with vorinostat enhances the antitumor activity of cisplatin in castration-resistant prostate cancer by inhibiting DNA damage repair pathway and detoxification of GSH.
The Prostate
Chang T,Lian Z,Ma S,Liang Z,Ma X,Wen X,Wang Y,Liu R
Sat Apr 01 00:00:00 UTC 2023
A Novel Myricetin Derivative with Anti-cancer Properties Induces Cell Cycle Arrest and Apoptosis in A549 Cells.
Biological & pharmaceutical bulletin
Zhou H,Xu L,Shi Y,Gu S,Wu N,Liu F,Huang Y,Qian Z,Xue W,Wang X,Chen F
Thu Jan 05 00:00:00 UTC 2023
Inhibition of miR-4640-5p alleviates pulmonary hypertension in chronic obstructive pulmonary disease patients by regulating nitric oxide synthase 1.
Respiratory research
Yang Z,Li P,Yuan Q,Wang X,Ma HH,Zhuan B
Fri Mar 24 00:00:00 UTC 2023
Cytotoxic Steroidal Saponins Containing a Rare Fructosyl from the Rhizomes of Paris polyphylla var. latifolia.
International journal of molecular sciences
Li TY,Du Y,Wang MC,Liu K,Liu Y,Cao Y,Wang YY,Chen WW,Qian XY,Qiu PC,Tang HF,Lu YY
Wed Apr 12 00:00:00 UTC 2023
Methylation-mediated silencing of EDN3 promotes cervical cancer proliferation, migration and invasion.
Frontiers in oncology
Zhu P,Li X,Liu Y,Xiong J,Yuan D,Chen Y,Luo L,Huang J,Wang B,Nie Q,Wang S,Dang L,Li S,Shu Y,Zhang W,Zhou H,Fan L,Li Q
Sun Feb 26 00:00:00 UTC 2023
Ginsenoside Compound K Ameliorates Osteoarthritis by Inhibiting the Chondrocyte Endoplasmic Reticulum Stress-Mediated IRE1α-TXNIP-NLRP3 Axis and Pyroptosis.
Journal of agricultural and food chemistry
Tian Y,Feng X,Zhou Z,Qin S,Chen S,Zhao J,Hou J,Liu D
Wed Jan 25 00:00:00 UTC 2023
Identification of MAP Kinase Kinase 3 as a protein target of myricetin in non-small cell lung cancer cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gu L,Li Z,Zhang X,Chen M,Zhang X
Mon May 01 00:00:00 UTC 2023
High-Dosage NMN Promotes Ferroptosis to Suppress Lung Adenocarcinoma Growth through the NAM-Mediated SIRT1-AMPK-ACC Pathway.
Cancers
Zhang M,Cui J,Chen H,Wang Y,Kuai X,Sun S,Tang Q,Zong F,Chen Q,Wu J,Wu S
Sun Apr 23 00:00:00 UTC 2023
The Protective Role of Scorias spongiosa Polysaccharide-Based Microcapsules on Intestinal Barrier Integrity in DSS-Induced Colitis in Mice.
Foods (Basel, Switzerland)
Xu Y,Feng H,Zhang Z,Zhang Q,Tang J,Zhou J,Wang Y,Peng W
Fri Feb 03 00:00:00 UTC 2023
Histone mRNA polyadenylation-mediated inflammation underlies various virus infections and cancers.
Journal of medical virology
Guo M,Zhu J,Hu Z,Wang Q,Songyang Z,Xiong Y
Thu Jun 01 00:00:00 UTC 2023
Protective Mechanism of Ethyl Gallate against Intestinal Ischemia-Reperfusion Injury in Mice by in Vivo and in Vitro Studies Based on Transcriptomics.
Chemistry & biodiversity
Fan S,Feng X,Li K,Li B,Diao Y
Sun Jan 01 00:00:00 UTC 2023
Bone microenvironment regulative hydrogels with ROS scavenging and prolonged oxygen-generating for enhancing bone repair.
Bioactive materials
Sun H,Xu J,Wang Y,Shen S,Xu X,Zhang L,Jiang Q
Thu Jun 01 00:00:00 UTC 2023
FSH-inhibited autophagy protects against oxidative stress in goat Sertoli cells through p62-Nrf2 pathway.
Theriogenology
Xi H,Hu Z,Han S,Liu X,Wang L,Hu J
Sun Jan 01 00:00:00 UTC 2023
AKR1B1 Represses Glioma Cell Proliferation through p38 MAPK-Mediated Bcl-2/BAX/Caspase-3 Apoptotic Signaling Pathways.
Current issues in molecular biology
Huang YK,Chang KC,Li CY,Lieu AS,Lin CL
Thu Apr 13 00:00:00 UTC 2023
Involvement of Galectin-3 in neurocognitive impairment in obstructive sleep apnea via regulating inflammation and oxidative stress through NLRP3.
Sleep medicine
Zong D,Liu X,Shen C,Liu T,Ouyang R
Sun Jan 01 00:00:00 UTC 2023
miR-484 mediates oxidative stress-induced ovarian dysfunction and promotes granulosa cell apoptosis via SESN2 downregulation.
Redox biology
Wang X,Yang J,Li H,Mu H,Zeng L,Cai S,Su P,Li H,Zhang L,Xiang W
Thu Jun 01 00:00:00 UTC 2023
SCAP contributes to embryonic angiogenesis by negatively regulating KISS-1 expression in mice.
Cell death & disease
Zheng G,Su Y,Wei L,Yao Y,Wang Y,Luo X,Wang X,Ruan XZ,Li D,Chen Y
Thu Apr 06 00:00:00 UTC 2023
Cholesterol-Lowering Activity of Vitisin A Is Mediated by Inhibiting Cholesterol Biosynthesis and Enhancing LDL Uptake in HepG2 Cells.
International journal of molecular sciences
Yuan Y,Zhu Y,Li Y,Li X,Jiao R,Bai W
Tue Feb 07 00:00:00 UTC 2023
Vitamin D receptor attenuate ischemia-reperfusion kidney injury via inhibiting ATF4.
Cell death discovery
Tang S,Wu X,Dai Q,Li Z,Yang S,Liu Y,Yi B,Wang J,Liao Q,Zhang W,Zhang H
Fri May 12 00:00:00 UTC 2023
Lyn attenuates sepsis-associated acute kidney injury by inhibition of phospho-STAT3 and apoptosis.
Biochemical pharmacology
Li N,Lin G,Zhang H,Sun J,Gui M,Liu Y,Li W,Zhan Z,Li Y,Pan S,Liu J,Tang J
Mon May 01 00:00:00 UTC 2023
Platycodin D suppresses proliferation, migration, and invasion of human glioblastoma cells through regulation of Skp2.
European journal of pharmacology
Li H,Ouyang J,Liu R
Mon Jun 05 00:00:00 UTC 2023
Exosomes incorporated with black phosphorus quantum dots attenuate retinal angiogenesis via disrupting glucose metabolism.
Materials today. Bio
Gui X,Zhang H,Zhang R,Li Q,Zhu W,Nie Z,Zhao J,Cui X,Hao W,Wen X,Shen W,Song H
Sat Apr 01 00:00:00 UTC 2023
HIV-1 Infection Promotes Cholesterol Aggregation by Inducing miR-33b-5p to Suppress ABCA1 Expression.
AIDS research and human retroviruses
You Q,Qiao J,Peng Q,Li W,Li J,Hu S,Shu Z,Sun B
Fri Sep 01 00:00:00 UTC 2023
The Bcr-Abl inhibitor DCC-2036 inhibits necroptosis and ameliorates osteoarthritis by targeting RIPK1 and RIPK3 kinases.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Piao L,Wu D,Rui C,Yang Y,Liu S,Liu J,Jin Z,Zhang H,Feng X,Bai L
Mon May 01 00:00:00 UTC 2023
Comparative analysis of two new zebrafish models: The cyp1a low-expression line and cyp1a knockout line under PAHs exposure.
Gene
Xie S,Feng Y,Zhou A,Lu Z
Thu Jun 15 00:00:00 UTC 2023
SLC17A9-PTHLH-EMT axis promotes proliferation and invasion of clear renal cell carcinoma.
iScience
Li W,Xu N,Meng X,Yuan H,Yu T,Miao Q,Yang H,Hai B,Xiao W,Zhang X
Fri Jan 20 00:00:00 UTC 2023
Effects of lactate on metabolism and differentiation of CD4+T cells.
Molecular immunology
Zhang YT,Xing ML,Fang HH,Li WD,Wu L,Chen ZP
Wed Feb 01 00:00:00 UTC 2023
Combined Application of Exosomes and FPR2 Agonist LXA4 in Controlling Fetal Membrane Inflammation and Promoting Fetal Membrane Tissue Repair.
Reproductive sciences (Thousand Oaks, Calif.)
Huang X,Liao J,Feng F,Chen S,Liao E,Li D,Dai X,Dong J,Shao Y
Thu Jun 01 00:00:00 UTC 2023
iPSC-based modeling of THD recapitulates disease phenotypes and reveals neuronal malformation.
EMBO molecular medicine
Tristán-Noguero A,Fernández-Carasa I,Calatayud C,Bermejo-Casadesús C,Pons-Espinal M,Colini Baldeschi A,Campa L,Artigas F,Bortolozzi A,Domingo-Jiménez R,Ibáñez S,Pineda M,Artuch R,Raya Á,García-Cazorla À,Consiglio A
Wed Mar 08 00:00:00 UTC 2023
A Recombinant Parathyroid Hormone-Related Peptide Locally Applied in Osteoporotic Bone Defect.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wang Y,Hu Y,Lan S,Chen Z,Zhang Y,Guo X,Cai L,Li J
Tue Aug 01 00:00:00 UTC 2023
Inhibition of Klf10 Attenuates Oxidative Stress-Induced Senescence of Chondrocytes via Modulating Mitophagy.
Molecules (Basel, Switzerland)
Shang J,Lin N,Peng R,Jiang N,Wu B,Xing B,Lin S,Xu X,Lu H
Tue Jan 17 00:00:00 UTC 2023
Hsa_circ_0001535 inhibits the proliferation and migration of ovarian cancer by sponging miR-593-3p, upregulating PTEN expression.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico
Han Y,Zheng Y,You J,Han Y,Lu X,Wang X,Shi C,Zhu W
Sun Oct 01 00:00:00 UTC 2023
PDCD6 Promotes Hepatocellular Carcinoma Cell Proliferation and Metastasis through the AKT/GSK3β/β-catenin Pathway.
Biomedical and environmental sciences : BES
Wen SY,Liu YT,Wei BY,Ma JQ,Chen YY
Mon Mar 20 00:00:00 UTC 2023
Deletion of LCMR1 in alveolar type II cells induces lethal impairment of lung structure and function in adult mice.
Journal of thoracic disease
Wang Y,Li C,Wu Z,Dai Y,Liu L,Chen L
Fri Mar 31 00:00:00 UTC 2023
Targeting ARF1-IQGAP1 interaction to suppress colorectal cancer metastasis and vemurafenib resistance.
Journal of advanced research
Hu HF,Gao GB,He X,Li YY,Li YJ,Li B,Pan Y,Wang Y,He QY
Fri Sep 01 00:00:00 UTC 2023
Blockage of glycolysis by targeting PFKFB3 suppresses the development of infantile hemangioma.
Journal of translational medicine
Yang K,Qiu T,Zhou J,Gong X,Zhang X,Lan Y,Zhang Z,Ji Y
Mon Feb 06 00:00:00 UTC 2023
PM2.5 induces autophagy-dependent ferroptosis by endoplasmic reticulum stress in SH-SY5Y cells.
Journal of applied toxicology : JAT
Guo CC,Xu ZM,Lyu Y,Li XH,Li ZF,He H,Tian FJ,Zheng JP
Sat Jul 01 00:00:00 UTC 2023
The molecular mechanism of miR-96-5p in the pathogenesis and treatment of polycystic ovary syndrome.
Translational research : the journal of laboratory and clinical medicine
Liu Y,Zhang S,Chen L,Huang X,Wang M,Ponikwicka-Tyszko D,Rahman NA,Wolczynski S,Yao B,Li X
Thu Jun 01 00:00:00 UTC 2023
Long noncoding RNA SNHG1 alleviates high glucose-induced vascular smooth muscle cells calcification/senescence by post-transcriptionally regulating Bhlhe40 and autophagy via Atg10.
Journal of physiology and biochemistry
Li S,Ni Y,Li C,Xiang Q,Zhao Y,Xu H,Huang W,Wang Y,Wang Y,Zhan J,Liu Y
Wed Feb 01 00:00:00 UTC 2023
Submersion and hypoxia inhibit alveolar epithelial Na+ transport through ERK/NF-κB signaling pathway.
Respiratory research
Zhou W,Hou Y,Yu T,Wang T,Ding Y,Nie H
Mon Apr 24 00:00:00 UTC 2023
Cuproptosis is involved in copper-induced hepatotoxicity in chickens.
The Science of the total environment
Zhong G,Li L,Li Y,Ma F,Liao J,Li Y,Zhang H,Pan J,Hu L,Tang Z
Sat Mar 25 00:00:00 UTC 2023
Cannabidiol alleviates neuroinflammation and attenuates neuropathic pain via targeting FKBP5.
Brain, behavior, and immunity
Wang X,Lin C,Jin S,Wang Y,Peng Y,Wang X
Sat Jul 01 00:00:00 UTC 2023
The role of selenoprotein M in nickel-induced pyroptosis in mice spleen tissue via oxidative stress.
Environmental science and pollution research international
Ma W,Liu Y,Xu L,Gai X,Sun Y,Qiao S,Liu P,Liu Q,Zhang Z
Wed Mar 01 00:00:00 UTC 2023
QiShenYiQi dripping pill alleviates myocardial ischemia-induced ferroptosis via improving mitochondrial dynamical homeostasis and biogenesis.
Journal of ethnopharmacology
Wu L,Fan Z,Gu L,Liu J,Cui Z,Yu B,Kou J,Li F
Tue May 23 00:00:00 UTC 2023
Effects of heat shock protein 90 on complement activation in myocardial ischemia/reperfusion injury after pioglitazone preconditioning.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University
Wang D,He S,Zhong G,Meng J,Bi Q,Tu R
Fri Dec 01 00:00:00 UTC 2023
Integrated Machine Learning and Bioinformatic Analyses Constructed a Network Between Mitochondrial Dysfunction and Immune Microenvironment of Periodontitis.
Inflammation
Chen H,Peng L,Wang Z,He Y,Zhang X
Sun Oct 01 00:00:00 UTC 2023
Polysaccharide-rich fractions from Enteromorpha prolifera improve hepatic steatosis and gut barrier integrity in high-fat diet-induced obese mice linking to modulation of gut microbiota.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zou T,Xie F,Liang P,Chen J,Wang Z,Du M,You J
Sun Jan 01 00:00:00 UTC 2023
A novel autophagy-related gene signature associated with prognosis and immune microenvironment in ovarian cancer.
Journal of ovarian research
Yang J,Wang C,Zhang Y,Cheng S,Wu M,Gu S,Xu S,Wu Y,Wang Y
Sat Apr 29 00:00:00 UTC 2023
Bach1 modulates AKT3 transcription to participate in hyperglycaemia-mediated EndMT in vascular endothelial cells.
Clinical and experimental pharmacology & physiology
Meng Z,Shen W,Yu L,Tong F,He H,Hu Y,Wu W,Liu J
Thu Jun 01 00:00:00 UTC 2023
GPR35 antagonist CID-2745687 attenuates anchorage-independent cell growth by inhibiting YAP/TAZ activity in colorectal cancer cells.
Frontiers in pharmacology
Otkur W,Liu X,Chen H,Li S,Ling T,Lin H,Yang R,Xia T,Qi H,Piao HL
Sun Apr 30 00:00:00 UTC 2023
Nicaraven protects against endotoxemia-induced inflammation and organ injury through modulation of AMPK/Sirt1 signaling in macrophages.
European journal of pharmacology
Zha D,Yang Y,Huang X,Wang Z,Lin H,Yang L,Xu L,Wu Y,Huang H,Wang Y,Xin Z,Wu X,Xiao YF,Li TS,Deng KY,Xin HB,Qian Y
Fri May 05 00:00:00 UTC 2023
Nanoengineered Gallium Ion Incorporated Formulation for Safe and Efficient Reversal of PARP Inhibition and Platinum Resistance in Ovarian Cancer.
Research (Washington, D.C.)
Li Y,Cen Y,Tu M,Xiang Z,Tang S,Lu W,Zhang H,Xu J
Mon Mar 20 00:00:00 UTC 2023
Degradation of Perineuronal Nets in the Cerebellar Interpositus Nucleus Ameliorated Social Deficits in Shank3-deficient Mice.
Neuroscience
Liu P,Zhao Y,Xiong W,Pan Y,Zhu M,Zhu X
Fri Feb 10 00:00:00 UTC 2023
Atf4 regulates angiogenic differences between alveolar bone and long bone macrophages by regulating M1 polarization, based on single-cell RNA sequencing, RNA-seq and ATAC-seq analysis.
Journal of translational medicine
Gu L,Wang Z,Gu H,Wang H,Liu L,Zhang WB
Tue Mar 14 00:00:00 UTC 2023
GPR109A controls neutrophil extracellular traps formation and improve early sepsis by regulating ROS/PAD4/Cit-H3 signal axis.
Experimental hematology & oncology
Guo W,Gong Q,Zong X,Wu D,Li Y,Xiao H,Song J,Zhang S,Fu S,Feng Z,Zhuang L
Tue Jan 31 00:00:00 UTC 2023
ROS-responsive magnesium-containing microspheres for antioxidative treatment of intervertebral disc degeneration.
Acta biomaterialia
Zhang T,Wang Y,Li R,Xin J,Zheng Z,Zhang X,Xiao C,Zhang S
Wed Mar 01 00:00:00 UTC 2023
Immunity-related long noncoding RNA WDFY3-AS2 inhibited cell proliferation and metastasis through Wnt/β-catenin signaling in oral squamous cell carcinoma.
Archives of oral biology
Lin X,Ding JM,Zheng XZ,Chen JG
Wed Mar 01 00:00:00 UTC 2023
Peripheral BDNF Regulates Somatosensory-Sympathetic Coupling in Brachial Plexus Avulsion-Induced Neuropathic Pain.
Neuroscience bulletin
Xian H,Guo H,Liu YY,Zhang JL,Hu WC,Yu MJ,Zhao R,Xie RG,Zhang H,Cong R
Fri Dec 01 00:00:00 UTC 2023
HSF4 promotes tumor progression of colorectal cancer by transactivating c-MET.
Molecular and cellular biochemistry
Zhang W,Zhang X,Cheng P,Yue K,Tang M,Li Y,Guo Q,Zhang Y
Mon May 01 00:00:00 UTC 2023
Nuclear factor Nrf2 promotes glycosidase OGG1 expression by activating the AKT pathway to enhance leukemia cell resistance to cytarabine.
The Journal of biological chemistry
Shang Q,Pan C,Zhang X,Yang T,Hu T,Zheng L,Cao S,Feng C,Hu X,Chai X,Wang J,Fang Q
Sun Jan 01 00:00:00 UTC 2023
African Swine Fever Virus L83L Negatively Regulates the cGAS-STING-Mediated IFN-I Pathway by Recruiting Tollip To Promote STING Autophagic Degradation.
Journal of virology
Cheng M,Kanyema MM,Sun Y,Zhao W,Lu Y,Wang J,Li X,Shi C,Wang J,Wang N,Yang W,Jiang Y,Huang H,Yang G,Zeng Y,Wang C,Cao X
Tue Feb 28 00:00:00 UTC 2023
An early-onset SLE patient with a novel paternal inherited BACH2 mutation.
Journal of clinical immunology
Zhou L,Sun G,Chen R,Chen J,Fang S,Xu Q,Tang W,Dai R,Zhang Z,An Y,Tang X,Zhao X
Tue Aug 01 00:00:00 UTC 2023
XBP1s gene of endoplasmic reticulum stress enhances proliferation and osteogenesis of human periodontal ligament cells.
Tissue & cell
Cui Z,Qin R,Feng J,Liu Y,Zhou X,Qin X,Li Y,Zhang Z,He X
Tue Aug 01 00:00:00 UTC 2023
Ginkgo biloba extract attenuates cisplatin-induced renal interstitial fibrosis by inhibiting the activation of renal fibroblasts through down-regulating the HIF-1α/STAT3/IL-6 pathway in renal tubular epithelial cells.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhang Y,Qin X,Yang Y,Li J,Li X,Zou X,Huang Z,Huang S
Sat Jul 01 00:00:00 UTC 2023
Effects of long-term resveratrol treatment in hypothalamic astrocyte cultures from aged rats.
Molecular and cellular biochemistry
Sovrani V,Bobermin LD,Santos CL,Brondani M,Gonçalves CA,Leipnitz G,Quincozes-Santos A
Thu Jun 01 00:00:00 UTC 2023
The effects of acupuncture on depression by regulating BDNF-related balance via lateral habenular nucleus BDNF/TrkB/CREB signaling pathway in rats.
Behavioural brain research
Tong T,Chen Y,Hao C,Shen J,Chen W,Cheng W,Yan S,Li J,Li Y,Gulizhaerkezi T,Zeng J,Meng X
Sat Aug 05 00:00:00 UTC 2023
Venoarterial extracorporeal membrane oxygenation improves survival in a rat model of acute myocardial infarction.
Artificial organs
Wei S,Cheng X,Li J,Zhai K,Huang J,Wang S,Li Y,Liu D
Sat Jul 01 00:00:00 UTC 2023
AQP8 may affect glioma proliferation and growth by regulating GSK-3β phosphorylation and nuclear transport of β-catenin.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience
Cai Z,Shen Z,Zhao J,Zhang H,Guo Z,Xia Q,Liang H,Liu J,Tan L,Sheng H,Zhu S
Thu Jun 01 00:00:00 UTC 2023
Microglial NLRP3 inflammasome activation mediates diabetes-induced depression-like behavior via triggering neuroinflammation.
Progress in neuro-psychopharmacology & biological psychiatry
Su WJ,Li JM,Zhang T,Cao ZY,Hu T,Zhong SY,Xu ZY,Gong H,Jiang CL
Wed Aug 30 00:00:00 UTC 2023
Glioma-derived LRIG3 interacts with NETO2 in tumor-associated macrophages to modulate microenvironment and suppress tumor growth.
Cell death & disease
Li Y,Wang W,Hou X,Huang W,Zhang P,He Y,Wang B,Duan Q,Mao F,Guo D
Fri Jan 13 00:00:00 UTC 2023
E2F1 combined with LINC01004 super-enhancer to promote hepatocellular carcinoma cell proliferation and metastasis.
Clinical epigenetics
Li J,Wang J,Wang Y,Zhao X,Su T
Tue Jan 31 00:00:00 UTC 2023
Staphylococci planktonic and biofilm environments differentially affect osteoclast formation.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Seebach E,Kraus FV,Elschner T,Kubatzky KF
Sat Jul 01 00:00:00 UTC 2023
TMEM41B Is an Interferon-Stimulated Gene That Promotes Pseudorabies Virus Replication.
Journal of virology
Li XQ,Zeng L,Liang DG,Qi YL,Yang GY,Zhong K,Chu BB,Wang J
Thu Jun 29 00:00:00 UTC 2023
Acupuncture exerts preventive effects in rats of chronic unpredictable mild stress: The involvement of inflammation in amygdala and brain-spleen axis.
Biochemical and biophysical research communications
Chen W,Chen Y,Cheng W,Li P,Shen J,Tong T,Lai L,Yan S,Huang Z,Li J,Huang S,Meng X
Sun Feb 26 00:00:00 UTC 2023
Metformin Collaborates with PINK1/Mfn2 Overexpression to Prevent Cardiac Injury by Improving Mitochondrial Function.
Biology
Ma Z,Liu Z,Li X,Zhang H,Han D,Xiong W,Zhou H,Yang X,Zeng Q,Ren H,Xu D
Tue Apr 11 00:00:00 UTC 2023
A novel feedback regulated loop of circRRM2-IGF2BP1-MYC promotes breast cancer metastasis.
Cancer cell international
Hao R,Zhang L,Si Y,Zhang P,Wang Y,Li B,Hu J,Qi Y
Sat Mar 25 00:00:00 UTC 2023
Promotion of intestinal epithelial cell apoptosis by enterotoxigenic Escherichia coli via PKA-mediated inhibition of mTORC1 activation.
Microbes and infection
Zheng Y,Wang G,Li R,Geng H,Lei X,Chen L,Wu S,Yao J,Deng L
Thu Jun 01 00:00:00 UTC 2023
Puerarin protects against sepsis-associated encephalopathy by inhibiting NLRP3/Caspase-1/GSDMD pyroptosis pathway and reducing blood-brain barrier damage.
European journal of pharmacology
Zhou S,Li Y,Hong Y,Zhong Z,Zhao M
Sat Apr 15 00:00:00 UTC 2023
Targeting lncRNA DDIT4-AS1 Sensitizes Triple Negative Breast Cancer to Chemotherapy via Suppressing of Autophagy.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Jiang T,Zhu J,Jiang S,Chen Z,Xu P,Gong R,Zhong C,Cheng Y,Sun X,Yi W,Yang J,Zhou W,Cheng Y
Thu Jun 01 00:00:00 UTC 2023
LncRNA SLC25A21-AS1 increases the chemosensitivity and inhibits the progression of ovarian cancer by upregulating the expression of KCNK4.
Functional & integrative genomics
Huang K,Chen X,Geng Z,Xiong X,Cong Y,Pan X,Liu S,Ge L,Xu J,Jia X
Thu Mar 30 00:00:00 UTC 2023
Long non-coding RNA ENST00000503625 is a potential prognostic biomarker and metastasis suppressor gene in prostate cancer.
Journal of cancer research and clinical oncology
Li Y,Fang Z,Ge S,Li J,Qu L,Shi X,Zhang W,Sun Y,Ren S,Wang L
Tue Aug 01 00:00:00 UTC 2023
The NLRP3 inflammasome recognizes alpha-2 and alpha-7.3 giardins and decreases the pathogenicity of Giardia duodenalis in mice.
Parasites & vectors
Zhao P,Li J,Li X,Dong J,Wang X,Zhang N,Li S,Sun M,Zhang X,Wang Z,Liang M,Li Y,Cao L,Gong P
Fri Mar 03 00:00:00 UTC 2023
APEX1 predicts poor prognosis of gallbladder cancer and affects biological properties of CD133+ GBC-SD cells via upregulating Jagged1.
Journal of Cancer
Wu Z,Liu Z,Sun Y,Yuan Y,Zou Q,Wen Y,Luo J,Liu R
Thu Jun 08 00:00:00 UTC 2023
Targeting the mitochondrial calcium uniporter inhibits cancer progression and alleviates cisplatin resistance in esophageal squamous cell carcinoma.
International journal of oncology
Miao Y,Wang X,Lai Y,Huang Y,Yin H,Meng X,Liu H,Hou R,Lin W,Zhang X,Zhang X,Chai BC,Zhang F,Guo L,Yang S
Sat Jul 01 00:00:00 UTC 2023
Spinal Cord Stimulation Attenuates Neural Remodeling, Inflammation, and Fibrosis After Myocardial Infarction.
Neuromodulation : journal of the International Neuromodulation Society
He Y,Sun Z,Jiang J,Yin X,Han J,Zhang Y,Zheng L
Sun Jan 01 00:00:00 UTC 2023
The Cytotoxic Cardiac Glycoside (-)-Cryptanoside A from the Stems of Cryptolepis dubia and Its Molecular Targets.
Journal of natural products
Ren Y,Kaweesa EN,Tian L,Wu S,Sydara K,Xayvue M,Moore CE,Soejarto DD,Cheng X,Yu J,Burdette JE,Kinghorn AD
Fri Jun 23 00:00:00 UTC 2023
Long non-coding RNA LINC00707, a prognostic marker, regulates cell proliferation, apoptosis, and EMT in esophageal squamous cell carcinoma.
American journal of translational research
Gao N,Jiang G,Gao Z,Cui M,Li J,Liu H,Fan T
Fri May 19 00:00:00 UTC 2023
Butyrate dictates ferroptosis sensitivity through FFAR2-mTOR signaling.
Cell death & disease
Wang G,Qin S,Chen L,Geng H,Zheng Y,Xia C,Yao J,Deng L
Tue Apr 25 00:00:00 UTC 2023
A Y374X TDP43 truncation leads to an altered metabolic profile in amyotrophic lateral sclerosis fibroblasts driven by pyruvate and TCA cycle intermediate alterations.
Frontiers in aging neuroscience
Allen SP,Al Sultan A,Kabucho Kibirige E,Tonkiss E,Hamer KJ,Castelli LM,Lin YH,Roscoe S,Stefanidis N,Mead RJ,Highley JR,Cooper-Knock J,Hautbergue GM,Heath PR,Kirby J,Shaw PJ
Wed May 31 00:00:00 UTC 2023
Heliangin acts as a covalent ligand of RPS2 that disrupts pre-rRNA metabolic processes in NPM1-mutated acute myeloid leukemia.
Acta pharmaceutica Sinica. B
Feng Y,Han Y,Hu A,Qu Y,Hu Y,Wu H,Wang X,He L
Wed Feb 01 00:00:00 UTC 2023
E7050 Suppresses the Growth of Multidrug-Resistant Human Uterine Sarcoma by Inhibiting Angiogenesis via Targeting of VEGFR2-Mediated Signaling Pathways.
International journal of molecular sciences
Huang TT,Chen CM,Lin SS,Lan YW,Cheng HC,Choo KB,Wang CC,Huang TH,Chong KY
Wed May 31 00:00:00 UTC 2023
Resveratrol and Dulaglutide ameliorate adiposity and liver dysfunction in rats with diet-induced metabolic syndrome: Role of SIRT-1 / adipokines / PPARγ and IGF-1.
Daru : journal of Faculty of Pharmacy, Tehran University of Medical Sciences
Shamardl HAMA,Ibrahim NA,Merzeban DH,Elamir AM,Golam RM,Elsayed AM
Thu Jun 01 00:00:00 UTC 2023
25-hydroxycholesterol inhibits human papillomavirus infection in cervical epithelial cells by perturbing cytoskeletal remodeling.
Journal of medical virology
Li B,Hua C,Tian P,Sha Y,Zhang L,Wang Q,Lu L,Jiang S,Sui L
Thu Jun 01 00:00:00 UTC 2023
IFN-α-2b Reduces Postoperative Arthrofibrosis in Rats by Inhibiting Fibroblast Proliferation and Migration through STAT1/p21 Signaling Pathway.
Mediators of inflammation
Liu Z,Fan Z,Wang R,Li X,Chen H,Wang J
Wed Mar 15 00:00:00 UTC 2023
Yishen Tongluo formula alleviates diabetic kidney disease through regulating Sirt6/TGF-β1/Smad2/3 pathway and promoting degradation of TGF-β1.
Journal of ethnopharmacology
Zhang X,Zhao L,Xiang S,Sun Y,Wang P,Chen JJ,Teo BS,Xie Z,Zhang Z,Xu J
Wed May 10 00:00:00 UTC 2023
Lovastatin inhibits erythroleukemia progression through KLF2-mediated suppression of MAPK/ERK signaling.
BMC cancer
Gao J,Hu J,Yu F,Wang C,Sheng D,Liu W,Hu A,Yu K,Xiao X,Kuang Y,Zacksenhaus E,Gajendran B,Ben-David Y
Tue Apr 04 00:00:00 UTC 2023
CXCL11 negatively regulated by MED19 favours antitumour immune infiltration in breast cancer.
Cytokine
Zhang X,Wu J,Hu C,Zheng X,Guo Z,Li L
Wed Feb 01 00:00:00 UTC 2023
Myricetin improves apoptosis after ischemic stroke via inhibiting MAPK-ERK pathway.
Molecular biology reports
Zhang L,Zhou T,Ji Q,He L,Lan Y,Ding L,Li L,Wang Z
Wed Mar 01 00:00:00 UTC 2023
Hypoxia Inhibits Cell Cycle Progression and Cell Proliferation in Brain Microvascular Endothelial Cells via the miR-212-3p/MCM2 Axis.
International journal of molecular sciences
Shi Q,Li S,Lyu Q,Zhang S,Bai Y,Ma J
Wed Feb 01 00:00:00 UTC 2023
VEZF1, destabilized by STUB1, affects cellular growth and metastasis of hepatocellular carcinoma by transcriptionally regulating PAQR4.
Cancer gene therapy
Shi X,Zhao P,Zhao G
Wed Feb 01 00:00:00 UTC 2023
Exogenous H2S Attenuates Hypertension by Regulating Renin Exocytosis under Hyperglycaemic and Hyperlipidaemic Conditions.
International journal of molecular sciences
Liu N,Li M,Liu S,Kang J,Chen L,Huang J,Wang Y,Chen H,Zhang W
Sat Jan 14 00:00:00 UTC 2023
Citrus peel ameliorates mucus barrier damage in HFD-fed mice.
The Journal of nutritional biochemistry
Han P,Yu Y,Zhang L,Ruan Z
Wed Feb 01 00:00:00 UTC 2023
m6A reader HNRNPA2B1 destabilization of ATG4B regulates autophagic activity, proliferation and olaparib sensitivity in breast cancer.
Experimental cell research
Zheng R,Yu Y,Lv L,Zhang Y,Deng H,Li J,Zhang B
Wed Mar 01 00:00:00 UTC 2023
BST2 regulated by the transcription factor STAT1 can promote metastasis, invasion and proliferation of oral squamous cell carcinoma via the AKT/ERK1/2 signaling pathway.
International journal of oncology
Shan F,Shen S,Wang X,Chen G
Sat Apr 01 00:00:00 UTC 2023
NAMPT inhibition reduces macrophage inflammation through the NAD+/PARP1 pathway to attenuate liver ischemia-reperfusion injury.
Chemico-biological interactions
Lu J,Wang M,Chen Y,Song H,Wen D,Tu J,Guo Y,Liu Z
Thu Jan 05 00:00:00 UTC 2023
Turnip crinkle virus-encoded suppressor of RNA silencing interacts with Arabidopsis SGS3 to enhance virus infection.
Molecular plant pathology
Liu L,Wang H,Fu Y,Tang W,Zhao P,Ren Y,Liu Z,Wu K,Zhang X
Wed Feb 01 00:00:00 UTC 2023
ZBTB7C m6A modification incurred by METTL3 aberration promotes osteosarcoma progression.
Translational research : the journal of laboratory and clinical medicine
An X,Wu W,Yang L,Dong J,Liu B,Guo J,Chen J,Guo B,Cao W,Jiang Q
Fri Sep 01 00:00:00 UTC 2023
Downregulation of lncRNA SLC7A11-AS1 decreased the NRF2/SLC7A11 expression and inhibited the progression of colorectal cancer cells.
PeerJ
Wang T,Liang S,Li Y,Wang X,Wang H,Guo J,Li M
Fri Apr 21 00:00:00 UTC 2023
Low dose TGF-β1 can improve vohwinkel syndrome by promoting the proliferation of keratinocytes.
Acta histochemica
Ling X,Dong S,Zhang L
Wed Feb 01 00:00:00 UTC 2023
FUNDC1 modulates mitochondrial defects and pancreatic β-cell dysfunction under lipotoxicity.
Biochemical and biophysical research communications
Tong B,Zhang Z,Li X,Liu J,Wang H,Song L,Feng J,Dai Z,Xu Y
Sun Sep 10 00:00:00 UTC 2023
Montelukast Inhibits Lung Cancer Cell Migration by Suppressing Cysteinyl Leukotriene Receptor 1 Expression In vitro.
Current pharmaceutical biotechnology
Chen Y,Zhang J,Wei S
Thu Jun 01 00:00:00 UTC 2023
Chinese herbal medicine alleviates the pathogenesis of polycystic ovary syndrome by improving oxidative stress and glucose metabolism via mitochondrial Sirtuin 3 signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhang Q,Ren J,Wang F,Li M,Pan M,Zhang H,Qu F
Sun Jan 01 00:00:00 UTC 2023
Mesenchymal stem cells ameliorate cisplatin-induced acute kidney injury via let-7b-5p.
Cell and tissue research
Wang SY,Xu Y,Hong Q,Chen XM,Cai GY
Mon May 01 00:00:00 UTC 2023
The Rab GTPase-binding protein EHBP1L1 and its interactors CD2AP/CIN85 negatively regulate the length of primary cilia via actin remodeling.
The Journal of biological chemistry
Iwano T,Sobajima T,Takeda S,Harada A,Yoshimura SI
Wed Mar 01 00:00:00 UTC 2023
Annao Pingchong decoction alleviate the neurological impairment by attenuating neuroinflammation and apoptosis in intracerebral hemorrhage rats.
Journal of ethnopharmacology
Guo C,Zhou X,Wang X,Wang H,Liu J,Wang J,Lin X,Lei S,Yang Y,Liu K,Long H,Zhou D
Wed Jun 28 00:00:00 UTC 2023
Identification of a chemoresistance-related prognostic gene signature by comprehensive analysis and experimental validation in pancreatic cancer.
Frontiers in oncology
Chen J,Liu Z,Wu Z,Li W,Tan X
Wed May 31 00:00:00 UTC 2023
Tryptophan Metabolism Acts as a New Anti-Ferroptotic Pathway to Mediate Tumor Growth.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu D,Liang CH,Huang B,Zhuang X,Cui W,Yang L,Yang Y,Zhang Y,Fu X,Zhang X,Du L,Gu W,Wang X,Yin C,Chai R,Chu B
Wed Feb 01 00:00:00 UTC 2023
Diagnostic value and underlying mechanism of nasal nitric oxide in eosinophilic chronic rhinosinusitis with nasal polyps.
Molecular immunology
Tang B,Tu J,Zhang M,Zhang Z,Yu J,Shen L,Luo Q,Ye J
Sat Jul 01 00:00:00 UTC 2023
AdipoRon mitigates tau pathology and restores mitochondrial dynamics via AMPK-related pathway in a mouse model of Alzheimer's disease.
Experimental neurology
Wang C,Chang Y,Zhu J,Wu Y,Jiang X,Zheng S,Li G,Ma R
Mon May 01 00:00:00 UTC 2023
Pharmacological vitamin C inhibits mTOR signaling and tumor growth by degrading Rictor and inducing HMOX1 expression.
PLoS genetics
Qin S,Wang G,Chen L,Geng H,Zheng Y,Xia C,Wu S,Yao J,Deng L
Wed Feb 01 00:00:00 UTC 2023
Jiang-Tang-San-Huang pill alleviates type 2 diabetes mellitus through modulating the gut microbiota and bile acids metabolism.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Tawulie D,Jin L,Shang X,Li Y,Sun L,Xie H,Zhao J,Liao J,Zhu Z,Cui H,Wen W
Mon May 01 00:00:00 UTC 2023
Shoutai Wan regulates glycolysis imbalance at the maternal-fetal interface in threatened abortion mice.
Journal of ethnopharmacology
Zhang L,Wang S,Ma Y,Song Y,Li D,Liang X,Hao Y,Jiang M,Lv J,Du H
Thu Aug 10 00:00:00 UTC 2023
Mechanism of the HIF-1α/VEGF/VEGFR-2 pathway in the proliferation and apoptosis of human haemangioma endothelial cells.
International journal of experimental pathology
Zhang W,Sun L,Gao H,Wang S
Sun Oct 01 00:00:00 UTC 2023
Effect of botanical drugs in improving symptoms of hypertensive nephropathy: Analysis of real-world data, retrospective cohort, network, and experimental assessment.
Frontiers in pharmacology
Huan JM,Ma XT,Li SY,Hu DQ,Chen HY,Wang YM,Su XY,Su WG,Wang YF
Tue Apr 25 00:00:00 UTC 2023
Comprehensive Genomic Profiling Identifies FAT1 as a Negative Regulator of EMT, CTCs, and Metastasis of Hepatocellular Carcinoma.
Journal of hepatocellular carcinoma
Huang ZL,Zhang PB,Zhang JT,Li F,Li TT,Huang XY
Wed Mar 15 00:00:00 UTC 2023
Pomegranate Peel Extract Sensitizes Hepatocellular Carcinoma Cells to Ionizing Radiation, Induces Apoptosis and Inhibits MAPK, JAK/STAT3, β-Catenin/NOTCH, and SOCS3 Signaling.
Integrative cancer therapies
Elbakry MMM,ElBakary NM,Hagag SA,Hemida EHA
Tue Jan 31 00:00:00 UTC 2023
Sirt1 protects against intervertebral disc degeneration induced by 1,25-dihydroxyvitamin D insufficiency in mice by inhibiting the NF-κB inflammatory pathway.
Journal of orthopaedic translation
Wang P,Yang C,Lu J,Ren Y,Goltzman D,Miao D
Mon May 01 00:00:00 UTC 2023
Brexpiprazole suppresses cell proliferation and de novo lipogenesis through AMPK/SREBP1 pathway in colorectal cancer.
Environmental toxicology
Li T,Liu X,Long X,Li Y,Xiang J,Lv Y,Zhao X,Shi S,Chen W
Sun Oct 01 00:00:00 UTC 2023
O-GlcNAcylation enhances Reticulon 2 protein stability and its promotive effects on gastric cancer progression.
Cellular signalling
Wang G,Xu Z,Sun J,Liu B,Ruan Y,Gu J,Song S
Tue Aug 01 00:00:00 UTC 2023
Circular RNA circ-TNRC6B inhibits the proliferation and invasion of esophageal squamous cell carcinoma cells by regulating the miR-452-5p/DAG1 axis.
Molecular oncology
Xu R,Ding P,Zhao X,Li Z,Liu F,Gu L,Zheng Y,Sang M,Meng L
Sat Jul 01 00:00:00 UTC 2023
TRIM7 inhibits encephalomyocarditis virus replication by activating interferon-β signaling pathway.
Veterinary microbiology
Li M,Yan J,Zhu H,Guo C,Jiang X,Gao Y,Liu X,Jiang P,Bai J
Thu Jun 01 00:00:00 UTC 2023
Maslinic Acid Ameliorates Myocardial Ischemia Reperfusion Injury-Induced Oxidative Stress via Activating Nrf2 and Inhibiting NF-[Formula: see text]B Pathways.
The American journal of Chinese medicine
Li Q,Li Z,Liu C,Xu M,Li T,Wang Y,Feng J,Yin X,Du X,Lu C
Mon Jun 19 00:00:00 UTC 2023
The Effect of an Essential Oil Blend on Growth Performance, Intestinal Health, and Microbiota in Early-Weaned Piglets.
Nutrients
Shao Y,Peng Q,Wu Y,Peng C,Wang S,Zou L,Qi M,Peng C,Liu H,Li R,Xiong X,Yin Y
Sat Jan 14 00:00:00 UTC 2023
BSHXF-medicated serum combined with ADSCs regulates the TGF-β1/Smad pathway to repair oxidatively damaged NPCs and its component analysis.
Journal of ethnopharmacology
Duan J,Li Z,Liu E,Long H,Chen L,Yang S
Wed Nov 15 00:00:00 UTC 2023
Astaxanthin promotes mitochondrial biogenesis and antioxidant capacity in chronic high-intensity interval training.
European journal of nutrition
Wang Y,Chen X,Baker JS,Davison GW,Xu S,Zhou Y,Bao X
Sat Apr 01 00:00:00 UTC 2023
α-Cyperone protects dopaminergic neurons and inhibits neuroinflammation in LPS-induced Parkinson's disease rat model via activating Nrf2/HO-1 and suppressing NF-κB signaling pathway.
International immunopharmacology
Huang B,Hu G,Zong X,Yang S,He D,Gao X,Liu D
Wed Feb 01 00:00:00 UTC 2023
cPKCγ Inhibits Caspase-9-Initiated Neuronal Apoptosis in an Ischemia Reperfusion Model In Vitro Through p38 MAPK-p90RSK-Bad Pathway.
Neurochemical research
Wei HP,Peng ZF,Shao KM,Zhang PH,Chen L,Hu JA,Chai H,Liu JM
Wed Feb 01 00:00:00 UTC 2023
Folate inhibits lipid deposition via the autophagy pathway in chicken hepatocytes.
Poultry science
Du ZQ,Pang YQ,Zhang Y,Wang L,Zhang R,Li H,Yang CX
Wed Feb 01 00:00:00 UTC 2023
Plumbagin Enhances the Anticancer Effects of PF Chemotherapy via Downregulation of the PI3K/AKT/mTOR/p70S6K Pathway in Human Tongue Squamous Cell Carcinoma.
Journal of oncology
Pan ST,Ye FF,Huang G,Qiu JX
Fri Feb 24 00:00:00 UTC 2023
NMR-Based Metabolic Profiling of the Effects of α-Ketoglutarate Supplementation on Energy-Deficient C2C12 Myotubes.
Molecules (Basel, Switzerland)
Li Y,Zhang S,Huang C,Lin D
Sun Apr 30 00:00:00 UTC 2023
CircRNA_0017076 acts as a sponge for miR-185-5p in the control of epithelial-to-mesenchymal transition of tubular epithelial cells during renal interstitial fibrosis.
Human cell
Zhang F,Zou H,Li X,Liu J,Xie Y,Chen M,Yu J,Wu X,Guo B
Mon May 01 00:00:00 UTC 2023
Liquiritigenin Attenuated Collagen-Induced Arthritis and Cardiac Complication via Inflammation and Fibrosis Inhibition in Mice.
Chemical & pharmaceutical bulletin
Ning X,Ni Y,Cao J,Zhang H
Tue Apr 04 00:00:00 UTC 2023
Lysosome-related exosome secretion mediated by miR-26b / Rab31 pathway was associated with the proliferation and migration of MCF-7 cells treated with BPA.
Ecotoxicology and environmental safety
Chen Y,Hu Z,Tang M,Huang F,Xiong Y,Ouyang D,He J,He S,Xian H,Hu D
Wed Mar 01 00:00:00 UTC 2023
Activation of autophagy inhibits the activation of NLRP3 inflammasome and alleviates sevoflurane-induced cognitive dysfunction in elderly rats.
BMC neuroscience
Zhou J,Zhang C,Fang X,Zhang N,Zhang X,Zhu Z
Sat Jan 28 00:00:00 UTC 2023
Hydrogen peroxide and iron ions can modulate lipid peroxidation, apoptosis, and the cell cycle, but do not have a significant effect on DNA double-strand break.
Biochemical and biophysical research communications
Yang H,Tai F,Wang T,Zheng X,Ge C,Qin Y,Fu H
Sun Apr 09 00:00:00 UTC 2023
Hypoxia disrupts the nasal epithelial barrier by inhibiting PTPN2 in chronic rhinosinusitis with nasal polyps.
International immunopharmacology
Zhang M,Xiong Y,Tu J,Tang B,Zhang Z,Yu J,Shen L,Luo Q,Ye J
Mon May 01 00:00:00 UTC 2023
PPARα Inhibits Astrocyte Inflammation Activation by Restoring Autophagic Flux after Transient Brain Ischemia.
Biomedicines
Luo D,Ye W,Chen L,Yuan X,Zhang Y,Chen C,Jin X,Zhou Y
Tue Mar 21 00:00:00 UTC 2023
Colocalization of Chikungunya Virus with Its Receptor MXRA8 during Cell Attachment, Internalization, and Membrane Fusion.
Journal of virology
Feng F,Bouma EM,Hu G,Zhu Y,Yu Y,Smit JM,Diamond MS,Zhang R
Wed May 31 00:00:00 UTC 2023
Proteome-Wide Analysis Reveals TFEB Targets for Establishment of a Prognostic Signature to Predict Clinical Outcomes of Colorectal Cancer.
Cancers
Huang Z,Zhu S,Han Z,Li C,Liang J,Wang Y,Zhang S,Zhang J
Wed Jan 25 00:00:00 UTC 2023
ABCG2 plays a central role in the dysregulation of 25-hydrovitamin D in Crohn's disease.
The Journal of nutritional biochemistry
Peng J,Song X,Zhu F,Zhang C,Xia J,Zou D,Liu J,Yin F,Yin L,Guo H,Liu J
Tue Aug 01 00:00:00 UTC 2023
Ameliorative effects of different doses of selenium against fluoride-triggered apoptosis and oxidative stress-mediated renal injury in rats through the activation of Nrf2/HO-1/NQO1 signaling pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Hu Y,Yan Z,He Y,Li Y,Li M,Li Y,Zhang D,Zhao Y,Ommati MM,Wang J,Huo M,Wang J
Sat Apr 01 00:00:00 UTC 2023
Effect of arsenic stress on the intestinal structural integrity and intestinal flora abundance of Cyprinus carpio.
Frontiers in microbiology
Shi X,Xu W,Che X,Cui J,Shang X,Teng X,Jia Z
Sun May 14 00:00:00 UTC 2023
Diosmetin Maintains Barrier Integrity by Reducing the Expression of ABCG2 in Colonic Epithelial Cells.
Journal of agricultural and food chemistry
Liu J,Fu L,Yin F,Yin L,Song X,Guo H,Liu J
Wed Jun 14 00:00:00 UTC 2023
Protective effect of total flavonoids of Engelhardia roxburghiana Wall. leaves against radiation-induced intestinal injury in mice and its mechanism.
Journal of ethnopharmacology
Wu S,Tian C,Tu Z,Guo J,Xu F,Qin W,Chang H,Wang Z,Hu T,Sun X,Ning H,Li Y,Gou W,Hou W
Sat Jul 15 00:00:00 UTC 2023
Ribosomal protein L32 enhances hepatocellular carcinoma progression.
Cancer medicine
Hou G,Lu Z,Jiang J,Yang X
Mon May 01 00:00:00 UTC 2023
Dual Antioxidant DH-217 Mitigated Cerebral Ischemia-Reperfusion Injury by Targeting IKKβ/Nrf2/HO-1 Signal Axis.
Neurochemical research
Shen M,Zheng Y,Li G,Chen Y,Huang L,Wu J,Hong C
Wed Feb 01 00:00:00 UTC 2023
LEM Domain Containing 1 Acts as a Novel Oncogene and Therapeutic Target for Triple-Negative Breast Cancer.
Cancers
Li X,Jiang S,Jiang T,Sun X,Guan Y,Fan S,Cheng Y
Fri May 26 00:00:00 UTC 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 UTC 2023
Endothelial progenitor cell-derived exosomes inhibit pulmonary artery smooth muscle cell in vitro proliferation and resistance to apoptosis by modulating the Mitofusin-2 and Ras-Raf-ERK1/2 signaling pathway.
European journal of pharmacology
Liu P,Gao S,Li Z,Pan S,Luo G,Ji Z
Thu Jun 15 00:00:00 UTC 2023
DHAV 3CD targets IRF7 and RIG-I proteins to block the type I interferon upstream signaling pathway.
Veterinary research
Xia X,Cheng A,Wang M,Ou X,Sun D,Zhang S,Mao S,Yang Q,Tian B,Wu Y,Huang J,Gao Q,Jia R,Chen S,Liu M,Zhao XX,Zhu D,Yu Y,Zhang L
Thu Jan 26 00:00:00 UTC 2023
Milk-derived exosomes carrying siRNA-KEAP1 promote diabetic wound healing by improving oxidative stress.
Drug delivery and translational research
Xiang X,Chen J,Jiang T,Yan C,Kang Y,Zhang M,Xiang K,Guo J,Jiang G,Wang C,Yang X,Chen Z
Fri Sep 01 00:00:00 UTC 2023
KLF2 is a clinical diagnostic and treatment biomarker of breast cancer.
Frontiers in cell and developmental biology
Li YZ,Xie J,Wang RQ,Gao XQ,Liu PJ,Liu J
Tue May 02 00:00:00 UTC 2023
Natural killer cells strengthen antitumor activity of cisplatin by immunomodulation and ameliorate cisplatin-induced side effects.
International urology and nephrology
Wang Z,Yang Z,Qu C,Li J,Wang X
Tue Aug 01 00:00:00 UTC 2023
The effect of allyl isothiocyanate on chondrocyte phenotype is matrix stiffness-dependent: Possible involvement of TRPA1 activation.
Frontiers in molecular biosciences
Che H,Shao Z,Ding J,Gao H,Liu X,Chen H,Cai S,Ge J,Wang C,Wu J,Hao Y
Tue Apr 04 00:00:00 UTC 2023
Intrathecal Injection of Botulinum Toxin Type A has an Analgesic Effect in Male Rats CCI Model by Inhibiting the Activation of Spinal P2X4R.
Neurochemical research
Zhou WM,Lei ZY,Shi YQ,Gong CY,Kai Z,Wei N,Wang LN,Zhang CJ,Zhang HH
Sun Oct 01 00:00:00 UTC 2023
Prognosis, immune microenvironment, and personalized treatment prediction in Rho GTPase-activating protein 4-mutant cervical cancer: Computer strategies for precision oncology.
Life sciences
Lu X,Ying Y,Zhang W,Zhang J,Li R,Wang W
Wed Feb 15 00:00:00 UTC 2023
A novel circular RNA circNLRP3 alleviated ricin toxin-induced TNF-α production through sponging miR-221-5p.
Toxicon : official journal of the International Society on Toxinology
Chen W,Chang Y,Sun C,Xu M,Dong M,Zhao N,Wang Y,Zhang J,Xu N,Liu W
Wed Mar 01 00:00:00 UTC 2023
Brown-fat-mediated tumour suppression by cold-altered global metabolism.
Nature
Seki T,Yang Y,Sun X,Lim S,Xie S,Guo Z,Xiong W,Kuroda M,Sakaue H,Hosaka K,Jing X,Yoshihara M,Qu L,Li X,Chen Y,Cao Y
Mon Aug 01 00:00:00 UTC 2022
A deafness-associated mitochondrial DNA mutation caused pleiotropic effects on DNA replication and tRNA metabolism.
Nucleic acids research
Meng F,Jia Z,Zheng J,Ji Y,Wang J,Xiao Y,Fu Y,Wang M,Ling F,Guan MX
Fri Sep 09 00:00:00 UTC 2022
Gypenosides ameliorate high-fat diet-induced non-alcoholic steatohepatitis via farnesoid X receptor activation.
Frontiers in nutrition
Li H,Xi Y,Liu H,Xin X
Tue Sep 13 00:00:00 UTC 2022
Ginsenoside Rg3 increases gemcitabine sensitivity of pancreatic adenocarcinoma via reducing ZFP91 mediated TSPYL2 destabilization.
Journal of ginseng research
Pan H,Yang L,Bai H,Luo J,Deng Y
Thu Sep 01 00:00:00 UTC 2022
Zika virus alters centrosome organization to suppress the innate immune response.
EMBO reports
Kodani A,Knopp KA,Di Lullo E,Retallack H,Kriegstein AR,DeRisi JL,Reiter JF
Mon Sep 05 00:00:00 UTC 2022
Astrocytic p75NTR expression provoked by ischemic stroke exacerbates the blood-brain barrier disruption.
Glia
Qin X,Wang J,Chen S,Liu G,Wu C,Lv Q,He X,Bai X,Huang W,Liao H
Sun May 01 00:00:00 UTC 2022
Effects of Xiao Chengqi Formula on Slow Transit Constipation by Assessing Gut Microbiota and Metabolomics Analysis in vitro and in vivo.
Frontiers in pharmacology
Zhou Q,Zhang D,Zhang H,Wan X,Hu B,Zou Q,Su D,Peng H,Huang D,Ren D
Wed Nov 29 00:00:00 UTC 2023
Circular RNA CircSHKBP1 accelerates the proliferation, invasion, angiogenesis, and stem cell-like properties via modulation of microR-766-5p/high mobility group AT-hook 2 axis in laryngeal squamous cell carcinoma.
Bioengineered
Chen F,Zhang H,Wang J
Sun May 01 00:00:00 UTC 2022
Daidzein alleviates doxorubicin-induced heart failure via the SIRT3/FOXO3a signaling pathway.
Food & function
Li H,Zhang M,Wang Y,Gong K,Yan T,Wang D,Meng X,Yang X,Chen Y,Han J,Duan Y,Zhang S
Thu Sep 22 00:00:00 UTC 2022
The protective effects of sulforaphane on high-fat diet-induced metabolic associated fatty liver disease in mice via mediating the FXR/LXRα pathway.
Food & function
Ma S,Pang X,Tian S,Sun J,Hu Q,Li X,Lu Y
Tue Dec 13 00:00:00 UTC 2022
TTK Protein Kinase promotes temozolomide resistance through inducing autophagy in glioblastoma.
BMC cancer
Yu J,Gao G,Wei X,Wang Y
Mon Jul 18 00:00:00 UTC 2022
Silencing LINC00491 Inhibits Pancreatic Cancer Progression through MiR-188-5p-induced Inhibition of ZFP91.
Journal of Cancer
Fang Z,Ruan B,Zhong M,Xiong J,Jiang Y,Song Z
Wed Apr 13 00:00:00 UTC 2022
Mitochondrial PKM2 deacetylation by procyanidin B2-induced SIRT3 upregulation alleviates lung ischemia/reperfusion injury.
Cell death & disease
Zhao J,Wang G,Han K,Wang Y,Wang L,Gao J,Zhao S,Wang G,Chen S,Luo A,Wu J,Wang G
Mon Jul 11 00:00:00 UTC 2022
Analysis of the expression and distribution of protein O-linked mannose β1,2-N-acetylglucosaminyltransferase 1 in the normal adult mouse brain.
Frontiers in neuroanatomy
Jiang H,Feng Y,He G,Liu Y,Li X
Wed Feb 01 00:00:00 UTC 2023
Circular RNA hsa_circ_0051040 Promotes Hepatocellular Carcinoma Progression by Sponging miR-569 and Regulating ITGAV Expression.
Cells
Ju L,Yao M,Lu R,Cao Y,Wang H,Yuan L,Xiao F,Shao J,Cai W,Chen L,Bian Z
Fri Nov 11 00:00:00 UTC 2022
GALNT3 protects against phosphate-induced calcification in vascular smooth muscle cells by enhancing active FGF23 and inhibiting the wnt/β-catenin signaling pathway.
Cellular signalling
Guo L,Wang Y,Li S,Zhou L,Li D
Thu Dec 01 00:00:00 UTC 2022
mascRNA promotes macrophage apoptosis, inhibits osteoclast differentiation and attenuates disease progression in a murine model of arthritis.
Biochemical and biophysical research communications
Wang X,Sun T,Mao X
Thu Jun 30 00:00:00 UTC 2022
Si-Wu Water Extracts Protect against Colonic Mucus Barrier Damage by Regulating Muc2 Mucin Expression in Mice Fed a High-Fat Diet.
Foods (Basel, Switzerland)
Ruan Z,Yu Y,Han P,Zhang L,Hu Z
Thu Aug 18 00:00:00 UTC 2022
RNF2 mediates pulmonary fibroblasts activation and proliferation by regulating mTOR and p16-CDK4-Rb1 signaling pathway.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Pan L,Hu Y,Qian C,Yao Y,Wang S,Shi W,Xu T
Tue Nov 01 00:00:00 UTC 2022
A Class I HDAC Inhibitor BG45 Alleviates Cognitive Impairment through the CaMKII/ITPKA/Ca2+ Signaling Pathway.
Pharmaceuticals (Basel, Switzerland)
Liu J,Zhang C,Wang J,Huang Y,Shen D,Hu Y,Chu H,Yu X,Zhang L,Ma H
Mon Nov 28 00:00:00 UTC 2022
Sphingomyelin-Sequestered Cholesterol Domain Recruits Formin-Binding Protein 17 for Constricting Clathrin-Coated Pits in Influenza Virus Entry.
Journal of virology
Tang B,Sun EZ,Zhang ZL,Liu SL,Liu J,Kusumi A,Hu ZH,Zeng T,Kang YF,Tang HW,Pang DW
Wed Mar 09 00:00:00 UTC 2022
Embryonic Deletion of TXNIP in GABAergic Neurons Enhanced Oxidative Stress in PV+ Interneurons in Primary Somatosensory Cortex of Aging Mice: Relevance to Schizophrenia.
Brain sciences
Xue T,Wang X,Hu Y,Cheng Y,Li H,Shi Y,Wang L,Yin D,Cui D
Sat Oct 15 00:00:00 UTC 2022
A homozygous PIWIL2 frameshift variant affects the formation and maintenance of human-induced pluripotent stem cell-derived spermatogonial stem cells and causes Sertoli cell-only syndrome.
Stem cell research & therapy
Wang X,Li Z,Qu M,Xiong C,Li H
Sat Sep 24 00:00:00 UTC 2022
IL-37 isoform D acts as an inhibitor of soluble ST2 to boost type 2 immune homeostasis in white adipose tissue.
Cell death discovery
Li C,Zhao M,Zhao M,Chen N,Guo Y,Du Y,Zhang Y,Cao B,Zhan B,Guo C,Li Y,Li Y,Shi Y,Zhu F,Zhang L,Wang Q
Tue Apr 05 00:00:00 UTC 2022
ANKRD37 inhibits trophoblast migration and invasion by regulating the NF-κB pathway in preeclampsia.
The journal of gene medicine
Tan W,Fu H,Zhou X,Duan Y,Yin N,Huang J,Liu X
Sun May 01 00:00:00 UTC 2022
Vam6 reduces iNKT cell function in tumor via modulating AMPK/mTOR pathways.
Frontiers in immunology
Bai S,Wu Q,Zhu S,Zhang Y,Chen X,Su M,Pan J,Li S,Yue T,Xu L,Xie D,Tian C,Zhao D,Li X,Hou J,Wang L,Fu S,Xue Y,Jiang A,Li D,Xu T,Tian Z,Zhou R,Zhang H,Bai L
Thu Feb 09 00:00:00 UTC 2023
Mitochondrial homeostasis regulates definitive endoderm differentiation of human pluripotent stem cells.
Cell death discovery
Lv J,Yi Y,Qi Y,Yan C,Jin W,Meng L,Zhang D,Jiang W
Thu Feb 17 00:00:00 UTC 2022
mTOR inhibition attenuates chemosensitivity through the induction of chemotherapy resistant persisters.
Nature communications
Liu Y,Azizian NG,Sullivan DK,Li Y
Thu Nov 17 00:00:00 UTC 2022
Ginsenoside Rg1 Inhibits STAT3 Expression by miR-15b-5p to Attenuate Lung Injury in Mice with Type 2 Diabetes Mellitus-Associated Pulmonary Tuberculosis.
Evidence-based complementary and alternative medicine : eCAM
Ma T,Mao X,Meng X,Wang Q
Wed Oct 19 00:00:00 UTC 2022
Identification of an autophagy-related 12-lncRNA signature and evaluation of NFYC-AS1 as a pro-cancer factor in lung adenocarcinoma.
Frontiers in genetics
Tong F,Xu L,Xu S,Zhang M
Thu Dec 22 00:00:00 UTC 2022
1α,25-Dihydroxyvitamin D3 Promotes Angiogenesis After Cerebral Ischemia Injury in Rats by Upregulating the TGF-β/Smad2/3 Signaling Pathway.
Frontiers in cardiovascular medicine
Zhang Y,Mu Y,Ding H,Du B,Zhou M,Li Q,Gong S,Zhang F,Geng D,Wang Y
Tue Apr 05 00:00:00 UTC 2022
Pseudorabies virus kinase UL13 phosphorylates H2AX to foster viral replication.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Ming X,Bo Z,Miao Y,Chen H,Bao C,Sun L,Xi R,Zhong Q,Zhao P,Jung YS,Qian Y
Tue Mar 01 00:00:00 UTC 2022
1,25(OH)2D3 alleviates LPS-induced preeclampsia-like rats impairment in the protective effect by TLR4/NF-kB pathway.
Placenta
Ma Y,Yang Y,Lv M,Zhang Y,He Q,Zhang Y,Su H,Deng X,Qian Y
Thu Dec 01 00:00:00 UTC 2022
p62 Promotes the Mitochondrial Localization of p53 through Its UBA Domain and Participates in Regulating the Sensitivity of Ovarian Cancer Cells to Cisplatin.
International journal of molecular sciences
Kong Q,Yan X,Cheng M,Jiang X,Xu L,Shen L,Yu H,Sun L
Fri Mar 18 00:00:00 UTC 2022
Correlation between CDK1 protein and CDK1 mRNA during oocyte maturation in mouse.
The International journal of developmental biology
Sun YT,Zhu AZ
Tue Oct 04 00:00:00 UTC 2022
IMP075 targeting ClpP for colon cancer therapy in vivo and in vitro.
Biochemical pharmacology
Zhang J,Luo B,Sui J,Qiu Z,Huang J,Yang T,Luo Y
Sat Oct 01 00:00:00 UTC 2022
Deoxythymidylate Kinase as a Promising Marker for Predicting Prognosis and Immune Cell Infiltration of Pan-cancer.
Frontiers in molecular biosciences
Lan T,Wang Y,Miao J,Guo H,Wang Z,Wang J,Zhang C,Yang P,Zhang Z,Dunmall LC,Wang Y
Sun Jul 31 00:00:00 UTC 2022
Zoledronic acid generates a spatiotemporal effect to attenuate osteoarthritis by inhibiting potential Wnt5a-associated abnormal subchondral bone resorption.
PloS one
Ding D,Wang L,Yan J,Zhou Y,Feng G,Ma L,Yang Y,Pei X,Jin Q
Mon Aug 01 00:00:00 UTC 2022
Sanguinarine protects against indomethacin-induced small intestine injury in rats by regulating the Nrf2/NF-κB pathways.
Frontiers in pharmacology
Lin XL,Shi YN,Cao YL,Tan X,Zeng YL,Luo ST,Li YM,Qin L,Xia BH,Fu RG,Lin LM,Li K,Cao D,Zeng JG,Liao DF
Sat Oct 29 00:00:00 UTC 2022
Ketamine Attenuates Airway Inflammation via Inducing Inflammatory Cells Apoptosis and Activating Nrf2 Pathway in a Mixed-Granulocytic Murine Asthma Model.
Drug design, development and therapy
Xiao S,Zhou Y,Wang Q,Yang D
Thu Jan 05 00:00:00 UTC 2023
Inhibition of CDK1 attenuates neuronal apoptosis and autophagy and confers neuroprotection after chronic spinal cord injury in vivo.
Journal of chemical neuroanatomy
Nie BX,Zhao G,Yuan XF,Yu LX,Zhang J,Yuan Y,Liu Y,Hu J,Song E,Zhou YC,Shu J
Sat Jan 01 00:00:00 UTC 2022
Dihydrotanshinone I Inhibits the Lung Metastasis of Breast Cancer by Suppressing Neutrophil Extracellular Traps Formation.
International journal of molecular sciences
Zhao H,Liang Y,Sun C,Zhai Y,Li X,Jiang M,Yang R,Li X,Shu Q,Kai G,Han B
Fri Dec 02 00:00:00 UTC 2022
Phosphorylation-Induced Ubiquitination and Degradation of PXR through CDK2-TRIM21 Axis.
Cells
Qin M,Xin Y,Bian Y,Yang X,Xi T,Xiong J
Thu Jan 13 00:00:00 UTC 2022
The effects of angiotensin I-converting enzyme inhibitory peptide VGINYW and the hydrolysate of α-lactalbumin on blood pressure, oxidative stress and gut microbiota of spontaneously hypertensive rats.
Food & function
Xie D,Shen Y,Su E,Du L,Xie J,Wei D
Mon Mar 07 00:00:00 UTC 2022
Paclitaxel Induces the Apoptosis of Prostate Cancer Cells via ROS-Mediated HIF-1α Expression.
Molecules (Basel, Switzerland)
Zhang Y,Tang Y,Tang X,Wang Y,Zhang Z,Yang H
Mon Oct 24 00:00:00 UTC 2022
NUPR1 contributes to radiation resistance by maintaining ROS homeostasis via AhR/CYP signal axis in hepatocellular carcinoma.
BMC medicine
Zhan Y,Zhang Z,Liu Y,Fang Y,Xie Y,Zheng Y,Li G,Liang L,Ding Y
Wed Oct 19 00:00:00 UTC 2022
LncRNA-CCAT1/miR-152-3p is involved in CSE-induced inflammation in HBE cells via regulating ERK signaling pathway.
International immunopharmacology
Zong D,Liu X,Li J,Long Y,Ouyang R,Chen Y
Mon Aug 01 00:00:00 UTC 2022
Identification and Validation of a Prognostic Signature Based on Methylation Profiles and Methylation-Driven Gene DAB2 as a Prognostic Biomarker in Differentiated Thyroid Carcinoma.
Disease markers
Ju G,Zhang L,Guo W,Wang H,Zhang X,Mu Z,Sun Y,Sun D,Diao H,Miao S,Chen Y,Xing T,Liang J,Lin Y
Fri May 26 00:00:00 UTC 2023
Protocatechuic aldehyde prevents ischemic injury by attenuating brain microvascular endothelial cell pyroptosis via lncRNA Xist.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Guo Y,Yang JH,He Y,Zhou HF,Wang Y,Ding ZS,Jin B,Wan HT
Sat Jan 01 00:00:00 UTC 2022
Ceria nanoparticles ameliorate renal fibrosis by modulating the balance between oxidative phosphorylation and aerobic glycolysis.
Journal of nanobiotechnology
Wang M,Zeng F,Ning F,Wang Y,Zhou S,He J,Li C,Wang C,Sun X,Zhang D,Xiao J,Hu P,Reilly S,Xin H,Xu X,Zhang X
Tue Jan 04 00:00:00 UTC 2022
Activation of PPARγ Protects Obese Mice from Acute Lung Injury by Inhibiting Endoplasmic Reticulum Stress and Promoting Mitochondrial Biogenesis.
PPAR research
Tang Y,Wei K,Liu L,Ma J,Wu S,Tang W
Tue Oct 11 00:00:00 UTC 2022
In situ forming and biocompatible hyaluronic acid hydrogel with reactive oxygen species-scavenging activity to improve traumatic brain injury repair by suppressing oxidative stress and neuroinflammation.
Materials today. Bio
Zhang D,Ren Y,He Y,Chang R,Guo S,Ma S,Guan F,Yao M
Wed Jun 01 00:00:00 UTC 2022
Long noncoding RNA lnc-H2AFV-1 promotes cell growth by regulating aberrant m6A RNA modification in head and neck squamous cell carcinoma.
Cancer science
Chen X,Liu Y,Sun D,Sun R,Wang X,Li M,Song N,Ying J,Guo T,Jiang Y
Wed Jun 01 00:00:00 UTC 2022
Network pharmacology-based and experimental identification of the effects of Renshen Yangrong decoction on myocardial infarction.
Frontiers in pharmacology
Zhao J,Xu X,Yang X
Fri Nov 18 00:00:00 UTC 2022
Resveratrol attenuates methylmercury-induced neurotoxicity by modulating synaptic homeostasis.
Toxicology and applied pharmacology
Wang W,Deng C,Chen F,Zhang L,Hu Y,Lu Q,Zhang A
Fri Apr 01 00:00:00 UTC 2022
Mitochondrial transcription factor B2 overexpression increases M2 macrophage infiltration via cytosolic mitochondrial DNA-stimulated Interleukin-6 secretion in ovarian cancer.
Bioengineered
Wu W,Zhou S,Liu T,Liang D
Sun May 01 00:00:00 UTC 2022
Pan-Cancer Analysis of the Immunological Role of PDIA5: A Potential Target for Immunotherapy.
Frontiers in immunology
Chen Y,He J,Chen R,Wang Z,Dai Z,Liang X,Wu W,Luo P,Zhang J,Peng Y,Zhang N,Liu Z,Zhang L,Zhang H,Cheng Q
Fri Aug 26 00:00:00 UTC 2022
Alterations in the expression level of visfatin in the lungs of piglets infected with PRRSV and its effect on PRRSV replication.
Microbial pathogenesis
Zhang ZW,Ansari AR,Dong L,Niu XY,Yang WJ,Li HZ,Xu FL,Yang KL,Song H
Tue Mar 01 00:00:00 UTC 2022
The NDUFV2 gene silencing inhibits the proliferation of two drug-resistant cancer cell lines.
Journal, genetic engineering & biotechnology
Liu L,Wang X,Li Y,Ma C,Shi Y,Li X,Chen J
Tue Apr 26 00:00:00 UTC 2022
The Translational Landscape Revealed the Sequential Treatment Containing ATRA plus PI3K/AKT Inhibitors as an Efficient Strategy for AML Therapy.
Pharmaceutics
Wang K,Ou Z,Deng G,Li S,Su J,Xu Y,Zhou R,Hu W,Chen F
Fri Oct 28 00:00:00 UTC 2022
Tumor-specific nitric oxide generator to amplify peroxynitrite based on highly penetrable nanoparticles for metastasis inhibition and enhanced cancer therapy.
Biomaterials
Shi M,Zhang J,Wang Y,Peng C,Hu H,Qiao M,Zhao X,Chen D
Fri Apr 01 00:00:00 UTC 2022
Low Sulfur Amino Acid, High Polyunsaturated Fatty Acid Diet Inhibits Breast Cancer Growth.
International journal of molecular sciences
Turchi R,Tortolici F,Benvenuto M,Punziano C,De Luca A,Rufini S,Faraonio R,Bei R,Lettieri-Barbato D,Aquilano K
Fri Dec 23 00:00:00 UTC 2022
Anti-inflammatory effect of Bifidobacterium animalis subsp. lactis A6 on DSS-induced colitis in mice.
Journal of applied microbiology
Wang H,Fan C,Zhao Z,Zhai Z,Hao Y
Thu Sep 01 00:00:00 UTC 2022
The "LLQY" Motif on SARS-CoV-2 Spike Protein Affects S Incorporation into Virus Particles.
Journal of virology
Du S,Xu W,Wang Y,Li L,Hao P,Tian M,Wang M,Li T,Wu S,Liu Q,Bai J,Qu X,Jin N,Zhou B,Liao M,Li C
Wed Mar 23 00:00:00 UTC 2022
High expression of B4GALT1 is associated with poor prognosis in acute myeloid leukemia.
Frontiers in genetics
Ren Z,Huang X,Lv Q,Lei Y,Shi H,Wang F,Wang M
Tue Jan 03 00:00:00 UTC 2023
Upregulation of Wilms' Tumor 1 in epicardial cells increases cardiac fibrosis in dystrophic mice.
Cell death and differentiation
Guo Z,Geng M,Huang Y,Han G,Jing R,Lin C,Zhang X,Zhang M,Fan G,Wang F,Yin H
Sat Oct 01 00:00:00 UTC 2022
Design, Synthesis, and Biological Evaluation of N14-Amino Acid-Substituted Tetrandrine Derivatives as Potential Antitumor Agents against Human Colorectal Cancer.
Molecules (Basel, Switzerland)
Wang YC,Zhang RH,Hu SC,Zhang H,Yang D,Zhang WL,Zhao YL,Cui DB,Li YJ,Pan WD,Liao SG,Zhou M
Thu Jun 23 00:00:00 UTC 2022
Stabilization of SAMHD1 by NONO is crucial for Ara-C resistance in AML.
Cell death & disease
Zhang F,Sun J,Tang X,Liang Y,Jiao Q,Yu B,Dai Z,Yuan X,Li J,Yan J,Zhang Z,Fan S,Wang M,Hu H,Zhang C,Lv XB
Fri Jul 08 00:00:00 UTC 2022
Transcriptome Profiling Analysis Identifies LCP1 as a Contributor for Chidamide Resistance in Gastric Cancer.
Pharmaceutical research
Bao W,Zhu Z,Gao Y,Chen J
Sun May 01 00:00:00 UTC 2022
Tetrahydroxy Stilbene Glucoside Alleviates Ischemic Stroke by Regulating Conformation-Dependent Intracellular Distribution of PKM2 for M2 Macrophage Polarization.
Journal of agricultural and food chemistry
Li M,Lu W,Meng Y,Zhang W,Wang F,Sun L,Xu Y
Wed Dec 14 00:00:00 UTC 2022
Phellinus linteus polysaccharides mediates acetaminophen-induced hepatotoxicity via activating AMPK/Nrf2 signaling pathways.
Aging
Zhao L,Zheng L,Li Z,Jin M,Wang Q,Cheng J,Li J,Feng H
Thu Sep 01 00:00:00 UTC 2022
Unveiling the anti-senescence effects and senescence-associated secretory phenotype (SASP) inhibitory mechanisms of Scutellaria baicalensis Georgi in low glucose-induced astrocytes based on boolean network.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Gao L,Yang WY,Qi H,Sun CJ,Qin XM,Du GH
Sun May 01 00:00:00 UTC 2022
A High-Fat Diet Induces Muscle Mitochondrial Dysfunction and Impairs Swimming Capacity in Zebrafish: A New Model of Sarcopenic Obesity.
Nutrients
Zou YY,Chen ZL,Sun CC,Yang D,Zhou ZQ,Xiao Q,Peng XY,Tang CF
Mon May 09 00:00:00 UTC 2022
LncRNA SENCR overexpression attenuated the proliferation, migration and phenotypic switching of vascular smooth muscle cells in aortic dissection via the miR-206/myocardin axis.
Nutrition, metabolism, and cardiovascular diseases : NMCD
Song Y,Wang T,Mu C,Gui W,Deng Y,Ma R
Wed Jun 01 00:00:00 UTC 2022
Isolation of five different primary cell types from a single sample of human skin.
STAR protocols
Kabacik S,Lowe D,Cohen H,Felton S,Spitzer J,Raj K
Fri Jun 17 00:00:00 UTC 2022
Carrier-free multifunctional nanomedicine for intraperitoneal disseminated ovarian cancer therapy.
Journal of nanobiotechnology
Huang X,Qiu M,Wang T,Li B,Zhang S,Zhang T,Liu P,Wang Q,Qian ZR,Zhu C,Wu M,Zhao J
Tue Feb 22 00:00:00 UTC 2022
PSMA1 mediates tumor progression and poor prognosis of gastric carcinoma by deubiquitinating and stabilizing TAZ.
Cell death & disease
Yang Q,Lu Y,Shangguan J,Shu X
Wed Nov 23 00:00:00 UTC 2022
Deletion of MTAP Highly Sensitizes Osteosarcoma Cells to Methionine Restriction With Recombinant Methioninase.
Cancer genomics & proteomics
Aoki Y,Tome Y,Han Q,Yamamoto J,Hamada K,Masaki N,Kubota Y,Bouvet M,Nishida K,Hoffman RM
Tue Apr 19 00:00:00 UTC 2022
XIST sponges miR-320d to promote chordoma progression by regulating ARF6.
Journal of bone oncology
Wang Y,Tang Z,Guo W
Mon Aug 01 00:00:00 UTC 2022
Transcriptome and Gut Microbiota Profiling Analysis of ANIT-Induced Cholestasis and the Effects of Da-Huang-Xiao-Shi Decoction Intervention.
Microbiology spectrum
Wang W,Jiang S,Xu C,Tang L,Liang Y,Zhao Y,Zhu G
Wed Dec 21 00:00:00 UTC 2022
Platycodon grandiflorum root fermentation broth reduces inflammation in a mouse IBD model through the AMPK/NF-κB/NLRP3 pathway.
Food & function
Wang Z,Li C,He X,Xu K,Xue Z,Wang T,Xu Z,Liu X
Mon Apr 04 00:00:00 UTC 2022
Pseudorabies Virus Inhibits Expression of Liver X Receptors to Assist Viral Infection.
Viruses
Wang Y,Li GL,Qi YL,Li LY,Wang LF,Wang CR,Niu XR,Liu TX,Wang J,Yang GY,Zeng L,Chu BB
Thu Mar 03 00:00:00 UTC 2022
Analysis of the absorbed constituents and mechanism of liquidambaris fructus extract on hepatocellular carcinoma.
Frontiers in pharmacology
Wang S,Yang XX,Li TJ,Zhao L,Bao YR,Meng XS
Sat Sep 17 00:00:00 UTC 2022
Effect of β-hydroxybutyrate on behavioral alterations, molecular and morphological changes in CNS of multiple sclerosis mouse model.
Frontiers in aging neuroscience
Sun W,Wen M,Liu M,Wang Q,Liu Q,Li L,Siebert HC,Loers G,Zhang R,Zhang N
Thu Dec 22 00:00:00 UTC 2022
Activation of RIPK2-mediated NOD1 signaling promotes proliferation and invasion of ovarian cancer cells via NF-κB pathway.
Histochemistry and cell biology
Zhang W,Wang Y
Tue Feb 01 00:00:00 UTC 2022
LINC00839 promotes colorectal cancer progression by recruiting RUVBL1/Tip60 complexes to activate NRF1.
EMBO reports
Liu X,Chen J,Zhang S,Liu X,Long X,Lan J,Zhou M,Zheng L,Zhou J
Mon Sep 05 00:00:00 UTC 2022
SiRNA-circFARSA-loaded porous silicon nanomaterials for pancreatic cancer treatment via inhibition of CircFARSA expression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Yuan H,Huang X,Li Q,Luo C,Lin C,Zhang S,Zhang Y,Yan Z,Du N,Liu Z,Jiang H,Chen B
Tue Mar 01 00:00:00 UTC 2022
Cytosine-phosphate-guanine oligodeoxynucleotides regulate the cell cycle, apoptosis, and steroidogenesis of mouse ovarian granulosa cells by targeting inhibin alpha (1 ~ 32) fragments.
In vitro cellular & developmental biology. Animal
Yang M,Feng J,Lan X,Tang Y,Zhang Y,Zhang H,Wang S
Tue Mar 01 00:00:00 UTC 2022
Angiotensin-(1-7) improves diabetes mellitus-induced erectile dysfunction in rats by regulating nitric oxide synthase levels.
Peptides
Xu Y,Zhang F,Li C,Hao H,Hao Y
Sun May 01 00:00:00 UTC 2022
Continuous theta-burst stimulation enhances and sustains neurogenesis following ischemic stroke.
Theranostics
Zong X,Gu J,Zhou S,Ding D,Hu Y,Tucker L,Huang Z,Geng D,Gao D
Tue Aug 16 00:00:00 UTC 2022
Channel HCN4 mutation R666Q associated with sporadic arrhythmia decreases channel electrophysiological function and increases protein degradation.
The Journal of biological chemistry
Wang H,Wu T,Huang Z,Huang J,Geng Z,Cui B,Yan Y,Zhang Y,Wang Y
Tue Nov 01 00:00:00 UTC 2022
HMGB1 downregulation in retinal pigment epithelial cells protects against diabetic retinopathy through the autophagy-lysosome pathway.
Autophagy
Feng L,Liang L,Zhang S,Yang J,Yue Y,Zhang X
Tue Feb 01 00:00:00 UTC 2022
HMGB1 downregulation in retinal pigment epithelial cells protects against diabetic retinopathy through the autophagy-lysosome pathway.
Autophagy
Feng L,Liang L,Zhang S,Yang J,Yue Y,Zhang X
Tue Feb 01 00:00:00 UTC 2022
Ferredoxin 1 is a cuproptosis-key gene responsible for tumor immunity and drug sensitivity: A pan-cancer analysis.
Frontiers in pharmacology
Yang L,Zhang Y,Wang Y,Jiang P,Liu F,Feng N
Tue Oct 11 00:00:00 UTC 2022
TLR4 Modulates Senescence and Paracrine Action in Placental Mesenchymal Stem Cells via Inhibiting Hedgehog Signaling Pathway in Preeclampsia.
Oxidative medicine and cellular longevity
Zhong Y,Zhang Y,Liu W,Zhao Y,Zou L,Liu X
Tue Jun 28 00:00:00 UTC 2022
LHPP suppresses gastric cancer progression via the PI3K/AKT/mTOR signaling pathway.
Journal of Cancer
Wang D,Li J,Li W
Sat Dec 10 00:00:00 UTC 2022
Sinapine Thiocyanate Inhibits the Proliferation and Mobility of Pancreatic Cancer Cells by Up-Regulating GADD45A.
Journal of Cancer
Wang J,Zeng Z,Lei S,Han J,Liao S,Zhang J,Wang L,Dong Y,Li H,Chen T
Tue Mar 15 00:00:00 UTC 2022
OCT4 induces EMT and promotes ovarian cancer progression by regulating the PI3K/AKT/mTOR pathway.
Frontiers in oncology
Xie W,Yu J,Yin Y,Zhang X,Zheng X,Wang X
Tue Aug 30 00:00:00 UTC 2022
Identification of Down-Regulated ADH1C is Associated With Poor Prognosis in Colorectal Cancer Using Bioinformatics Analysis.
Frontiers in molecular biosciences
Li M,Liu Z,Song J,Wang T,Wang H,Wang Y,Guo J
Sat Mar 19 00:00:00 UTC 2022
Injectable hydrogel with nucleus pulposus-matched viscoelastic property prevents intervertebral disc degeneration.
Journal of orthopaedic translation
Jia H,Lin X,Wang D,Wang J,Shang Q,He X,Wu K,Zhao B,Peng P,Wang H,Wang D,Li P,Yang L,Luo Z,Yang L
Tue Mar 01 00:00:00 UTC 2022
Hsa_circ_0008234 facilitates proliferation of cutaneous squamous cell carcinoma through targeting miR-127-5p to regulate ADCY7.
Archives of dermatological research
Cai L,Wang Y,Wu J,Wu G
Mon Aug 01 00:00:00 UTC 2022
Drp1 knockdown represses apoptosis of rat retinal endothelial cells by inhibiting mitophagy.
Acta histochemica
Wu H,Li G,Chen W,Luo W,Yang Z,You Z,Zou Y
Sat Jan 01 00:00:00 UTC 2022
Mitochondria-Targeting Polymer Micelle of Dichloroacetate Induced Pyroptosis to Enhance Osteosarcoma Immunotherapy.
ACS nano
Jin J,Yuan P,Yu W,Lin J,Xu A,Xu X,Lou J,Yu T,Qian C,Liu B,Song J,Li L,Piao Y,Xie T,Shen Y,Tao H,Tang J
Tue Jul 26 00:00:00 UTC 2022
Exosomal circRELL1 serves as a miR-637 sponge to modulate gastric cancer progression via regulating autophagy activation.
Cell death & disease
Sang H,Zhang W,Peng L,Wei S,Zhu X,Huang K,Yang J,Chen M,Dang Y,Zhang G
Thu Jan 13 00:00:00 UTC 2022
Heat stress-induced mucosal barrier dysfunction is potentially associated with gut microbiota dysbiosis in pigs.
Animal nutrition (Zhongguo xu mu shou yi xue hui)
Xia B,Wu W,Fang W,Wen X,Xie J,Zhang H
Tue Mar 01 00:00:00 UTC 2022
Mitochondrial fission induces immunoescape in solid tumors through decreasing MHC-I surface expression.
Nature communications
Lei X,Lin H,Wang J,Ou Z,Ruan Y,Sadagopan A,Chen W,Xie S,Chen B,Li Q,Wang J,Lin H,Zhu X,Yuan X,Tian T,Lv X,Fu S,Zhu X,Zhou J,Pan G,Xia X,Tannous BA,Ferrone S,Fan S,Li J
Wed Jul 06 00:00:00 UTC 2022
Recurrent Hypoglycemia Impaired Vascular Function in Advanced T2DM Rats by Inducing Pyroptosis.
Oxidative medicine and cellular longevity
Luo M,Hu Y,Lv D,Xie L,Yang S,Zuo D,Xue Y,He A
Wed Aug 03 00:00:00 UTC 2022
Icaritin attenuates 6-OHDA-induced MN9D cell damage by inhibiting oxidative stress.
PeerJ
Zhou X,Huang N,Hou X,Zhu L,Xie Y,Ba Z,Luo Y
Fri Jan 13 00:00:00 UTC 2023
KLF16 Downregulates the Expression of Tumor Suppressor Gene TGFBR3 to Promote Bladder Cancer Proliferation and Migration.
Cancer management and research
Chen X,Wang P,Ou T,Li J
Sun May 01 00:00:00 UTC 2022
The Novel Competing Endogenous Long Noncoding RNA SM2 Regulates Gonadotropin Secretion in the Hu Sheep Anterior Pituitary by Targeting the Oar-miR-16b/TGF-β/SMAD2 Signaling Pathway.
Cells
Wan Z,Yang H,Chen P,Wang Z,Cai Y,Yao X,Wang F,Zhang Y
Mon Mar 14 00:00:00 UTC 2022
Sinensetin attenuates IL-1β-induced cartilage damage and ameliorates osteoarthritis by regulating SERPINA3.
Food & function
Liu Z,Liu R,Wang R,Dai J,Chen H,Wang J,Li X
Mon Oct 03 00:00:00 UTC 2022
Exogenous TIPE2 Inhibit TAK1 to Improve Inflammation and Neuropathic Pain Induced by Sciatic Nerve Injury Through Inactivating NF-κB and JNK.
Neurochemical research
Sun X,Li X,Zhou Y,Wang Y,Liu X
Sat Oct 01 00:00:00 UTC 2022
Relationship between the expression of ARHGAP25 and RhoA in non-small cell lung cancer and vasculogenic mimicry.
BMC pulmonary medicine
Shi F,Wu J,Jia Q,Li K,Li W,Shi Y,Wang Y,Wu S
Fri Oct 07 00:00:00 UTC 2022
Theaflavin-3,3'-Digallate Protects Cartilage from Degradation by Modulating Inflammation and Antioxidant Pathways.
Oxidative medicine and cellular longevity
Teng Y,Jin Z,Ren W,Lu M,Hou M,Zhou Q,Wang W,Yang H,Zou J
Tue Jul 19 00:00:00 UTC 2022
Aucubin promoted neuron functional recovery by suppressing inflammation and neuronal apoptosis in a spinal cord injury model.
International immunopharmacology
Xiao S,Zhong N,Yang Q,Li A,Tong W,Zhang Y,Yao G,Wang S,Liu J,Liu Z
Sat Oct 01 00:00:00 UTC 2022
Carbon dots derived from folic acid attenuates osteoarthritis by protecting chondrocytes through NF-κB/MAPK pathway and reprogramming macrophages.
Journal of nanobiotechnology
Jin Y,Zhang Q,Qin X,Liu Z,Li Z,Zhong X,Xia L,He J,Fang B
Thu Nov 03 00:00:00 UTC 2022
Klotho alleviates NLRP3 inflammasome-mediated neuroinflammation in a temporal lobe epilepsy rat model by activating the Nrf2 signaling pathway.
Epilepsy & behavior : E&B
Xiang T,Luo X,Ye L,Huang H,Wu Y
Tue Mar 01 00:00:00 UTC 2022
Impact prediction of translocation of the mitochondrial outer membrane 70 as biomarker in Alzheimer's disease.
Frontiers in aging neuroscience
Cao X,Chen Y,Sang X,Xu S,Xie Z,Zhu Z,Wang P,Bi J,Xu L
Tue Nov 22 00:00:00 UTC 2022
Injectable multifunctional CMC/HA-DA hydrogel for repairing skin injury.
Materials today. Bio
Cui L,Li J,Guan S,Zhang K,Zhang K,Li J
Tue Mar 01 00:00:00 UTC 2022
Integrated Network Pharmacology and Experimental Verification to Explore the Molecular Mechanism of Hedysarum Multijugum Maxim-Curcumae Rhizoma Herb Pair for Treating Non-Small Cell Lung Cancer.
Frontiers in oncology
Hu S,Ge M,Zhang S,Jiang M,Hu K,Gao L
Sat May 07 00:00:00 UTC 2022
FOXA1 is a transcriptional activator of Odf2/Cenexin and regulates primary ciliation.
Scientific reports
Czerny CC,Borschel A,Cai M,Otto M,Hoyer-Fender S
Mon Dec 12 00:00:00 UTC 2022
Melatonin prevents cyclophosphamide-induced primordial follicle loss by inhibiting ovarian granulosa cell apoptosis and maintaining AMH expression.
Frontiers in endocrinology
Feng J,Ma WW,Li HX,Pei XY,Deng SL,Jia H,Ma WZ
Tue Aug 23 00:00:00 UTC 2022
Zinc Alleviates Arsenic-Induced Inflammation and Apoptosis in the Head Kidney of Common Carp by Inhibiting Oxidative Stress and Endoplasmic Reticulum Stress.
Biological trace element research
Xing P,Zhang Y,Chi Q,Li S
Sun May 01 00:00:00 UTC 2022
Progranulin improves neural development via the PI3K/Akt/GSK-3β pathway in the cerebellum of a VPA-induced rat model of ASD.
Translational psychiatry
Wang L,Chen J,Hu Y,Liao A,Zheng W,Wang X,Lan J,Shen J,Wang S,Yang F,Wang Y,Li Y,Chen D
Tue Mar 22 00:00:00 UTC 2022
Enhancement of the functionality of attenuating acute lung injury by a microemulsion formulation with volatile oil of Angelicae Sinensis Radix and Ligusticum Chuanxiong Rhizoma encapsulated.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang L,Yan Q,Zhang W,Li X,Zhang X,Du S,Hua X,Lin J,Shu G,Peng G,Tan Z,Fu H
Thu Dec 01 00:00:00 UTC 2022
Citrinin-Induced Hepatotoxicity in Mice Is Regulated by the Ca2+/Endoplasmic Reticulum Stress Signaling Pathway.
Toxins
Wu D,Yang C,Yang M,Wu Y,Mao Y,Zhou X,Wang J,Yuan Z,Wu J
Wed Apr 06 00:00:00 UTC 2022
β-elemene Isopropanolamine Derivative LXX-8250 Induces Apoptosis Through Impairing Autophagic Flux via PFKFB4 Repression in Melanoma Cells.
Frontiers in pharmacology
Jalal S,Zhang T,Deng J,Wang J,Xu T,Zhang T,Zhai C,Yuan R,Teng H,Huang L
Tue Aug 30 00:00:00 UTC 2022
Old targets, new strategy: Apigenin-7-O-β-d-(-6″-p-coumaroyl)-glucopyranoside prevents endothelial ferroptosis and alleviates intestinal ischemia-reperfusion injury through HO-1 and MAO-B inhibition.
Free radical biology & medicine
Feng YD,Ye W,Tian W,Meng JR,Zhang M,Sun Y,Zhang HN,Wang SJ,Wu KH,Liu CX,Liu SY,Cao W,Li XQ
Sun May 01 00:00:00 UTC 2022
Global Phosphoproteomics Unveils Kinase-Regulated Networks in Systemic Lupus Erythematosus.
Molecular & cellular proteomics : MCP
Meng S,Li T,Wang T,Li D,Chen J,Li H,Cai W,Zeng Z,Liu D,Tang D,Hong X,Dai Y
Thu Dec 01 00:00:00 UTC 2022
Role of non-neuronal cholinergic system in the early stage response of epithelial-mesenchymal transformation related markers in A549 cells induced by coal particles.
Heliyon
Wu MY,Shi XC,Shan J,Wang R,Wang Y,Li J,Tian DN,Xu HM
Tue Nov 01 00:00:00 UTC 2022
Protection of CAPE-pNO2 Against Chronic Myocardial Ischemia by the TGF-Β1/Galectin-3 Pathway In Vivo and In Vitro.
Inflammation
Wan Q,Zhang L,Zhou Q,Han Y,Li Z,Li B
Wed Jun 01 00:00:00 UTC 2022
Effect of Thymoquinone on Renal Damage Induced by Hyperlipidemia in LDL Receptor-Deficient (LDL-R-/-) Mice.
BioMed research international
Li W,Zhang H,Zhang L,Zhang T,Ding H
Tue Jul 19 00:00:00 UTC 2022
Geniposide attenuates spermatogenic dysfunction via inhibiting endoplasmic reticulum stress in male mice.
Chemico-biological interactions
Chi YN,Ye RJ,Yang JM,Hai DM,Liu N,Ren JW,Du J,Lan XB,Yu JQ,Ma L
Sat Oct 01 00:00:00 UTC 2022
Intracameral Injection of AAV-DJ.COMP-ANG1 Reduces the IOP of Mice by Reshaping the Trabecular Outflow Pathway.
Investigative ophthalmology & visual science
Qiao Y,Sun Z,Tan C,Lai J,Sun X,Chen J
Thu Dec 01 00:00:00 UTC 2022
Salvianolic acid A regulates pyroptosis of endothelial cells via directly targeting PKM2 and ameliorates diabetic atherosclerosis.
Frontiers in pharmacology
Zhu J,Chen H,Le Y,Guo J,Liu Z,Dou X,Lu D
Sat Nov 26 00:00:00 UTC 2022
lncRNA SNHG26 promoted the growth, metastasis, and cisplatin resistance of tongue squamous cell carcinoma through PGK1/Akt/mTOR signal pathway.
Molecular therapy oncolytics
Jiang Q,Wang Z,Qi Q,Li J,Xin Y,Qiu J
Thu Mar 17 00:00:00 UTC 2022
Canagliflozin Attenuates Hepatic Steatosis and Atherosclerosis Progression in Western Diet-Fed ApoE-Knockout Mice.
Drug design, development and therapy
Zuo Q,Zhang G,He L,Ma S,Ma H,Zhai J,Wang Z,Zhang T,Wang Y,Guo Y
Wed Dec 14 00:00:00 UTC 2022
Conditional knockout of ASK1 in microglia/macrophages attenuates epileptic seizures and long-term neurobehavioural comorbidities by modulating the inflammatory responses of microglia/macrophages.
Journal of neuroinflammation
Zhang Y,Wang Z,Wang R,Xia L,Cai Y,Tong F,Gao Y,Ding J,Wang X
Mon Aug 08 00:00:00 UTC 2022
Self-construction of actin networks through phase separation-induced abLIM1 condensates.
Proceedings of the National Academy of Sciences of the United States of America
Yang S,Liu C,Guo Y,Li G,Li D,Yan X,Zhu X
Tue Jul 19 00:00:00 UTC 2022
Inhibition of calpain9 attenuates peritoneal dialysis-related peritoneal fibrosis.
Frontiers in pharmacology
Li F,Wang Y,Tian J,Zhou Z,Yin W,Qin X,Wang H,Zeng T,Li A,Jiang J
Wed Dec 21 00:00:00 UTC 2022
Probiotic-fermented Pueraria lobata (Willd.) Ohwi alleviates alcoholic liver injury by enhancing antioxidant defense and modulating gut microbiota.
Journal of the science of food and agriculture
Zhao W,Peng D,Li W,Chen S,Liu B,Huang P,Wu J,Du B,Li P
Thu Dec 01 00:00:00 UTC 2022
Molecular Hydrogen Reduces Electromagnetic Pulse-Induced Male Rat Reproductive System Damage in a Rodent Model.
Oxidative medicine and cellular longevity
Ma L,Hao W,Feng WB,Cao LC,Qin LN,Wang Y,Liu MH,Wang NZ,Gao F,Guo JM,Du H,Yan HL
Fri Oct 07 00:00:00 UTC 2022
FTO-dependent N(6)-Methyladenosine regulates the progression of endometriosis via the ATG5/PKM2 Axis.
Cellular signalling
Wang H,Liang Z,Gou Y,Li Z,Cao Y,Jiao N,Tan J,Yu Y,Zhang Z
Sat Oct 01 00:00:00 UTC 2022
Altered gene expression in human brain microvascular endothelial cells in response to the infection of influenza H1N1 virus.
Animal diseases
Higazy D,Lin X,Xie T,Wang K,Gao X,Cui M
Wed Nov 09 00:00:00 UTC 2022
Panax notoginseng saponins protect PC12 cells against Aβ induced injury via promoting parkin-mediated mitophagy.
Journal of ethnopharmacology
Jiang Y,Li H,Huang P,Li S,Li B,Huo L,Zhong J,Pan Z,Li Y,Xia X
Tue Mar 01 00:00:00 UTC 2022
RNA-seq transcriptome profiling of liver regeneration in mice identifies the miR-34b-5p/phosphoinositide-dependent protein kinase 1 axis as a potential target for hepatocyte proliferation.
Biochemical and biophysical research communications
Lei X,Dai X,Wang Q,Long R,Xiang Z,Li H,Long Z,Zhang C,Zhu Z
Sun Oct 30 00:00:00 UTC 2022
Exposure to melamine cyanuric acid in adult mice caused motor activity and skeletal muscle energy metabolism disorder.
Physiology & behavior
Wang S,Zuo X,Xu B,Yu Q,An Z,Feng D,Feng X
Thu Dec 01 00:00:00 UTC 2022
Extracellular vesicles rich in HAX1 promote angiogenesis by modulating ITGB6 translation.
Journal of extracellular vesicles
You B,Pan S,Gu M,Zhang K,Xia T,Zhang S,Chen W,Xie H,Fan Y,Yao H,Cheng T,Zhang P,Liu D,You Y
Sun May 01 00:00:00 UTC 2022
A positive feedback loop of the TAZ/β-catenin axis promotes Helicobacter pylori-associated gastric carcinogenesis.
Frontiers in microbiology
Xu X,Shu C,Wu X,Ouyang Y,Cheng H,Zhou Y,Wang H,He C,Xie C,He X,Hong J,Lu N,Ge Z,Zhu Y,Li N
Wed Jan 11 00:00:00 UTC 2023
Diterpenoids from the whole plant of Euphorbia wallichii and their protective effects on H2O2-induced BV-2 microglial cells injury.
Bioorganic chemistry
Wang Y,Zhu M,Liang J,Zhang N,Sun D,Li H,Chen L
Tue Nov 01 00:00:00 UTC 2022
Transmissible Gastroenteritis Virus Nucleocapsid Protein Interacts with Na+/H+ Exchanger 3 To Reduce Na+/H+ Exchanger Activity and Promote Piglet Diarrhea.
Journal of virology
Niu Z,Xu S,Zhang Y,Kan Z,Zhang J,Liu X,Zhang S,Zou H,Song Z
Wed Nov 23 00:00:00 UTC 2022
Qingfei oral liquid alleviates RSV-induced lung inflammation by promoting fatty-acid-dependent M1/M2 macrophage polarization via the Akt signaling pathway.
Journal of ethnopharmacology
An L,Lu M,Xu W,Chen H,Feng L,Xie T,Shan J,Wang S,Lin L
Tue Nov 15 00:00:00 UTC 2022
Silibinin improves L-cell mass and function through an estrogen receptor-mediated antioxidative mechanism.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang J,Zhang L,Cao H,Shi X,Zhang X,Gao Z,Ikeda K,Yan T,Jia Y,Xu F
Sun May 01 00:00:00 UTC 2022
Phosphorylation-mediated PI3K-Art signalling pathway as a therapeutic mechanism in the hydrogen-induced alleviation of brain injury in septic mice.
Journal of cellular and molecular medicine
Bai Y,Li L,Dong B,Ma W,Chen H,Yu Y
Tue Nov 01 00:00:00 UTC 2022
HJURP Promotes Malignant Progression and Mediates Sensitivity to Cisplatin and WEE1-inhibitor in Serous Ovarian Cancer.
International journal of biological sciences
Dou Z,Qiu C,Zhang X,Yao S,Zhao C,Wang Z,Chu R,Chen J,Chen Z,Li R,Wang K,Liu P,Liu C,Song K,Kong B
Fri Apr 15 00:00:00 UTC 2022
Morchella esculenta mushroom polysaccharide attenuates diabetes and modulates intestinal permeability and gut microbiota in a type 2 diabetic mice model.
Frontiers in nutrition
Rehman AU,Siddiqui NZ,Farooqui NA,Alam G,Gul A,Ahmad B,Asim M,Khan AI,Xin Y,Zexu W,Song Ju H,Xin W,Lei S,Wang L
Tue Oct 25 00:00:00 UTC 2022
HDAC9 Contributes to Serous Ovarian Cancer Progression through Regulating Epithelial-Mesenchymal Transition.
Biomedicines
Xu L,Wang J,Liu B,Fu J,Zhao Y,Yu S,Shen L,Yan X,Su J
Thu Feb 03 00:00:00 UTC 2022
Inhibition of Metastatic Hepatocarcinoma by Combined Chemotherapy with Silencing VEGF/VEGFR2 Genes through a GalNAc-Modified Integrated Therapeutic System.
Molecules (Basel, Switzerland)
Li X,Wang X,Liu N,Wang Q,Hu J
Thu Mar 24 00:00:00 UTC 2022
Effects of bamboo leaf extract intervention on the growth performance, antioxidant capacity, and hepatic apoptosis in suckling piglets.
Journal of animal science
Yu G,Ji S,Yun Y,Cheng K,Zhang H,Jia P,Wang T,Zhang L
Fri Jul 01 00:00:00 UTC 2022
SIRT3-AMPK signaling pathway as a protective target in endothelial dysfunction of early sepsis.
International immunopharmacology
Yu H,Liu Q,Chen G,Huang L,Luo M,Lv D,Luo S
Sun May 01 00:00:00 UTC 2022
Self-Assembly of a Multifunction DNA Tetrahedron for Effective Delivery of Aptamer PL1 and Pcsk9 siRNA Potentiate Immune Checkpoint Therapy for Colorectal Cancer.
ACS applied materials & interfaces
Guo W,Gao H,Li H,Ge S,Zhang F,Wang L,Shi H,Han A
Wed Jul 20 00:00:00 UTC 2022
An in vitro senescence model of gingival epithelial cell induced by hydrogen peroxide treatment.
Odontology
Giri S,Takada A,Paudel D,Yoshida K,Furukawa M,Kuramitsu Y,Matsushita K,Abiko Y,Furuichi Y
Sat Jan 01 00:00:00 UTC 2022
Gliko BMSC: A potential strategy of treatment for renal fibrosis.
Regenerative therapy
Shi L,Gao X,Bi Y,Li M,Sun H,Tian X,Bi W
Wed Jun 01 00:00:00 UTC 2022
A specific EMC subunit supports Dengue virus infection by promoting virus membrane fusion essential for cytosolic genome delivery.
PLoS pathogens
Bagchi P,Speckhart K,Kennedy A,Tai AW,Tsai B
Fri Jul 01 00:00:00 UTC 2022
LGALS3BP in Microglia Promotes Retinal Angiogenesis Through PI3K/AKT Pathway During Hypoxia.
Investigative ophthalmology & visual science
Zhao C,Liu Y,Meng J,Wang X,Liu X,Li W,Zhou Q,Xiang J,Li N,Hou S
Fri Jul 08 00:00:00 UTC 2022
Pyroptosis participates in PM2.5-induced air-blood barrier dysfunction.
Environmental science and pollution research international
Jia R,Wei M,Zhang X,Du R,Sun W,Wang L,Song L
Mon Aug 01 00:00:00 UTC 2022
FLI1 accelerates leukemogenesis through transcriptional regulation of pyruvate kinase-L/R and other glycolytic genes.
Medical oncology (Northwood, London, England)
Sheng D,Chen B,Wang C,Xiao X,Hu A,Liu W,Kuang Y,Sample KM,Zacksenhaus E,Gajendran B,Pan W,Ben-David Y
Sat Dec 31 00:00:00 UTC 2022
Minocycline Pretreatment Prevents Blood-Brain Barrier Disruption in Septic Rats.
The Journal of surgical research
Yang G,Cao Y,Wang P,Mei L,Chen J,Lu W
Sun May 01 00:00:00 UTC 2022
PM2.5 induced lung injury through upregulating ROS-dependent NLRP3 Inflammasome-Mediated Pyroptosis.
Immunobiology
Ren F,Xu J,Zhang J,Xu X,Huang L,Sun W,Li R,Li F
Sun May 01 00:00:00 UTC 2022
DL-3-n-butylphthalide imparts neuroprotection via Nrf2/SIRT3 pathway in a mouse model of vascular dementia.
Brain research
Gao L,Guo X,Liu S,Sun Q,Qin X,Lv P,Hu M,Xu J,Dong Y
Tue Mar 15 00:00:00 UTC 2022
D-tagatose protects against oleic acid-induced acute respiratory distress syndrome in rats by activating PTEN/PI3K/AKT pathway.
Frontiers in immunology
Huang J,Wang B,Tao S,Hu Y,Wang N,Zhang Q,Wang C,Chen C,Gao B,Cheng X,Li Y
Tue Oct 04 00:00:00 UTC 2022
IL6 Induces mtDNA Leakage to Affect the Immune Escape of Endometrial Carcinoma via cGAS-STING.
Journal of immunology research
Zeng X,Li X,Zhang Y,Cao C,Zhou Q
Tue Jun 14 00:00:00 UTC 2022
The BDNF-TrkB-CREB Signalling Pathway Is Involved in Bisphenol S-Induced Neurotoxicity in Male Mice by Regulating Methylation.
Toxics
Li YZ,Wu ZY,Zhu BQ,Wang YX,Kan YQ,Zeng HC
Sat Jul 23 00:00:00 UTC 2022
HIF-1α inhibition attenuates severity of Achilles tendinopathy by blocking NF-κB and MAPK pathways.
International immunopharmacology
Jiao X,Zhang Y,Li W,Zhou X,Chu W,Li Y,Wang Z,Sun X,Xu C,Gan Y
Sun May 01 00:00:00 UTC 2022
Hypoxia-Inducible Factor 1α Stability Modified by Glutaredoxin-1 in Necrotizing Enterocolitis.
The Journal of surgical research
Zhang Y,Zhang X,Tian B,Deng Q,Guo C
Thu Dec 01 00:00:00 UTC 2022
The Role of Apoptosis and Autophagy in the Hypothalamic-Pituitary-Adrenal (HPA) Axis after Traumatic Brain Injury (TBI).
International journal of molecular sciences
Taheri S,Karaca Z,Mehmetbeyoglu E,Hamurcu Z,Yilmaz Z,Dal F,Çınar V,Ulutabanca H,Tanriverdi F,Unluhizarci K,Rassoulzadegan M,Kelestimur F
Sat Dec 10 00:00:00 UTC 2022
Overexpression of LINC00551 promotes autophagy-dependent ferroptosis of lung adenocarcinoma via upregulating DDIT4 by sponging miR-4328.
PeerJ
Peng X,Yang R,Peng W,Zhao Z,Tu G,He B,Cai Q,Shi S,Yin W,Yu F,Tao Y,Wang X
Tue Dec 27 00:00:00 UTC 2022
The underlying rationality of Chinese medicine herb pair Coptis chinensis and Dolomiaea souliei: From the perspective of metabolomics and intestinal function.
Journal of ethnopharmacology
Wang T,Zhang C,Li H,Zhou R,Ye X,Yang Y,He K
Tue May 10 00:00:00 UTC 2022
Arachidonic Acid in Follicular Fluid of PCOS Induces Oxidative Stress in a Human Ovarian Granulosa Tumor Cell Line (KGN) and Upregulates GDF15 Expression as a Response.
Frontiers in endocrinology
Ma Y,Zheng L,Wang Y,Gao Y,Xu Y
Thu Jun 02 00:00:00 UTC 2022
Reversion of methionine addiction of osteosarcoma cells to methionine independence results in loss of malignancy, modulation of the epithelial-mesenchymal phenotype and alteration of histone-H3 lysine-methylation.
Frontiers in oncology
Aoki Y,Han Q,Tome Y,Yamamoto J,Kubota Y,Masaki N,Obara K,Hamada K,Wang JD,Inubushi S,Bouvet M,Clarke SG,Nishida K,Hoffman RM
Tue Nov 22 00:00:00 UTC 2022
Acetyl CoA synthase 2 potentiates ATG5-induced autophagy against neuronal apoptosis after subarachnoid hemorrhage.
Journal of molecular histology
He W,Zhou X,Wu Q,Zhou L,Zhang Z,Zhang R,Deng C,Zhang X
Fri Apr 01 00:00:00 UTC 2022
Necrostatin-1 Against Sevoflurane-Induced Cognitive Dysfunction Involves Activation of BDNF/TrkB Pathway and Inhibition of Necroptosis in Aged Rats.
Neurochemical research
Yin C,Zhang Q,Zhao J,Li Y,Yu J,Li W,Wang Q
Fri Apr 01 00:00:00 UTC 2022
Immobilization of peroxisome proliferator-activated receptor gamma and the application in screening modulators of the receptor from herbal medicine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Hou Z,Jin Y,Li Y,Fan H,Xiao C,Li Q,Zhang Y
Tue Feb 01 00:00:00 UTC 2022
α1 -Adrenoceptors activate the NLRP3 inflammasome through downregulation of Kir2.1 in cardiac inflammation.
Experimental physiology
Chen L,Wang W,Peng X,Liu L,Zhang A,Li X,Ma K,Wang L
Wed Jun 01 00:00:00 UTC 2022
POLD2 is activated by E2F1 to promote triple-negative breast cancer proliferation.
Frontiers in oncology
Zhang Z
Tue Sep 20 00:00:00 UTC 2022
The inhibition of PGAM5 suppresses seizures in a kainate-induced epilepsy model via mitophagy reduction.
Frontiers in molecular neuroscience
Zhong F,Gan Y,Song J,Zhang W,Yuan S,Qin Z,Wu J,Lü Y,Yu W
Wed Jan 11 00:00:00 UTC 2023
A dynamic fluorescence probed glycolysis suppression process in HeLa cells treated with trichostatin A.
The Analyst
Zhang F,Shao C,Wu Y,Zhao W,Jing X,Fang C,Huang Q
Mon Nov 07 00:00:00 UTC 2022
Improvement of tube formation model of cell: Application for acute hypoxia in in vitro study of angiogenesis.
Microvascular research
Li F,Gou X,Xu D,Han D,Hou K,Fang W,Li Y
Tue Mar 01 00:00:00 UTC 2022
Improvement of tube formation model of cell: Application for acute hypoxia in in vitro study of angiogenesis.
Microvascular research
Li F,Gou X,Xu D,Han D,Hou K,Fang W,Li Y
Tue Mar 01 00:00:00 UTC 2022
Improvement of tube formation model of cell: Application for acute hypoxia in in vitro study of angiogenesis.
Microvascular research
Li F,Gou X,Xu D,Han D,Hou K,Fang W,Li Y
Tue Mar 01 00:00:00 UTC 2022
Improvement of tube formation model of cell: Application for acute hypoxia in in vitro study of angiogenesis.
Microvascular research
Li F,Gou X,Xu D,Han D,Hou K,Fang W,Li Y
Tue Mar 01 00:00:00 UTC 2022
Effect and mechanism of TFEB on pyroptosis in HK-2 cells induced by high glucose.
Biochemical and biophysical research communications
Han N,Wang Z,Luo H,Chi Y,Zhang T,Wang B,Li Y
Sat Jun 25 00:00:00 UTC 2022
Dexamethasone Alleviates Myocardial Injury in a Rat Model of Acute Myocardial Infarction Supported by Venoarterial Extracorporeal Membrane Oxygenation.
Frontiers in public health
Cheng X,Zhang R,Wei S,Huang J,Zhai K,Li Y,Gao B
Mon Aug 08 00:00:00 UTC 2022
Map of Enteropathogenic Escherichia coli Targets Mitochondria and Triggers DRP-1-Mediated Mitochondrial Fission and Cell Apoptosis in Bovine Mastitis.
International journal of molecular sciences
Li Y,Zhu Y,Chu B,Liu N,Chen S,Wang J,Zou Y
Thu Apr 28 00:00:00 UTC 2022
Sodium selenite attenuates zearalenone-induced apoptosis through inhibition of endoplasmic reticulum stress in goat trophoblast cells.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Xiong Y,Li B,Li J,Jin E,He S
Mon Aug 01 00:00:00 UTC 2022
Decreased tubulin-binding cofactor B was involved in the formation disorder of nascent astrocyte processes by regulating microtubule plus-end growth through binding with end-binding proteins 1 and 3 after chronic alcohol exposure.
Frontiers in cellular neuroscience
Zheng Y,Yang M,Chen X,Zhang G,Wan S,Zhang B,Huo J,Liu H
Fri Nov 18 00:00:00 UTC 2022
1,25-D3 attenuates cerebral ischemia injury by regulating mitochondrial metabolism via the AMPK/AKT/GSK3β pathway.
Frontiers in aging neuroscience
Li Y,Li X,Xu S,Zhao Y,Pang M,Zhang X,Wang X,Wang Y
Fri Nov 04 00:00:00 UTC 2022
LncRNA BIRF Promotes Brain Ischemic Tolerance Induced By Cerebral Ischemic Preconditioning Through Upregulating GLT-1 via Sponging miR-330-5p.
Molecular neurobiology
Li S,Zhang L,Lin J,Su A,Liu X,Zhang J,Xian X,Hu Y,Li W,Sun S,Zhang M
Fri Jul 01 00:00:00 UTC 2022
Glucolipotoxicity induces endothelial cell dysfunction by activating autophagy and inhibiting autophagic flow.
Diabetes & vascular disease research
Liu Y,Xiang H,Xiong W,Ouyang J,Liu H,Zhao S,Xiao J,Li J,Shu Z,Wang X,Liu H,Zhang J,Fan J,Li Y,Chen S,Lu H
Tue May 17 00:00:00 UTC 2022
Hydrogen sulphide attenuates neuronal apoptosis of substantia nigra by re-establishing autophagic flux via promoting leptin signalling in a 6-hydroxydopamine rat model of Parkinson's disease.
Clinical and experimental pharmacology & physiology
Jiang W,Zou W,Hu M,Tian Q,Xiao F,Li M,Zhang P,Chen YJ,Jiang JM
Sat Jan 01 00:00:00 UTC 2022
Hydrogen gas promotes apoptosis of lung adenocarcinoma A549 cells through X-linked inhibitor of apoptosis and baculoviral inhibitor of apoptosis protein repeat-containing 3.
Journal of cancer research and therapeutics
Zhang Y,Chen G,Yan Z,Wang L,Wang D
Thu Sep 01 00:00:00 UTC 2022
Hydrogen gas promotes apoptosis of lung adenocarcinoma A549 cells through X-linked inhibitor of apoptosis and baculoviral inhibitor of apoptosis protein repeat-containing 3.
Journal of cancer research and therapeutics
Zhang Y,Chen G,Yan Z,Wang L,Wang D
Thu Sep 01 00:00:00 UTC 2022
Hydrogen gas promotes apoptosis of lung adenocarcinoma A549 cells through X-linked inhibitor of apoptosis and baculoviral inhibitor of apoptosis protein repeat-containing 3.
Journal of cancer research and therapeutics
Zhang Y,Chen G,Yan Z,Wang L,Wang D
Thu Sep 01 00:00:00 UTC 2022
Hydrogen gas promotes apoptosis of lung adenocarcinoma A549 cells through X-linked inhibitor of apoptosis and baculoviral inhibitor of apoptosis protein repeat-containing 3.
Journal of cancer research and therapeutics
Zhang Y,Chen G,Yan Z,Wang L,Wang D
Thu Sep 01 00:00:00 UTC 2022
Actin-Binding LIM 1 (ABLIM1) Inhibits Glioblastoma Progression and Serves as a Novel Prognostic Biomarker.
Disease markers
Liu D,Wang X,Liu Y,Li C,Zhang Z,Lv P
Tue Jan 03 00:00:00 UTC 2023
Cyclosporin A up-regulated thromboxane A2 receptor through activation of MAPK and NF-κB pathways in rat mesenteric artery.
European journal of pharmacology
Wang C,Han L,Wang T,Wang Y,Liu J,Wang B,Xu CB
Tue Jul 05 00:00:00 UTC 2022
Mucin O-glycan-microbiota axis orchestrates gut homeostasis in a diarrheal pig model.
Microbiome
Xia B,Zhong R,Wu W,Luo C,Meng Q,Gao Q,Zhao Y,Chen L,Zhang S,Zhao X,Zhang H
Wed Aug 31 00:00:00 UTC 2022
The Aurora Kinase Inhibitor TAK901 Inhibits Glioblastoma Growth by Blocking SREBP1-Mediated Lipid Metabolism.
Cancers
Zhan X,Qiu R,He Y,Zhao Z,Huang M,Liu Q,Zhi F,Long W
Fri Nov 25 00:00:00 UTC 2022
Trichostatin D as a Novel KLF2 Activator Attenuates TNFα-Induced Endothelial Inflammation.
International journal of molecular sciences
Lei L,Chen M,Wang C,Jiang X,Li Y,Wang W,Li S,Zhao L,Sheng R,Han J,Zhang Y,Chen Y,Yan B,Wu Y,Yu L,Si S,Xu Y
Thu Nov 03 00:00:00 UTC 2022
Influenza virus NS1 interacts with 14-3-3ε to antagonize the production of RIG-I-mediated type I interferons.
Virology
Wang T,Wei F,Jiang Z,Song J,Li C,Liu J
Thu Sep 01 00:00:00 UTC 2022
Senescent Phenotype of Astrocytes Leads to Activation of BV2 Microglia and N2a Neuronal Cells Death.
Molecules (Basel, Switzerland)
Zhang W,Yang X,Liu J,Pan Y,Zhang M,Chen L
Mon Sep 12 00:00:00 UTC 2022
Bisdemethoxycurcumin Alleviates Dextran Sodium Sulfate-Induced Colitis via Inhibiting NLRP3 Inflammasome Activation and Modulating the Gut Microbiota in Mice.
Antioxidants (Basel, Switzerland)
Zhang J,Li Q,Zhang X,Chen Y,Lu Y,Wang X,Zhang L,Wang T
Fri Oct 07 00:00:00 UTC 2022
Downregulation of HINFP induces senescence-associated secretory phenotype to promote metastasis in a non-cell-autonomous manner in bladder cancer.
Oncogene
Zheng X,Liu Z,Zhong J,Zhou L,Chen J,Zheng L,Li Z,Zhang R,Pan J,Wu Y,Liu Z,Kang T
Fri Jul 01 00:00:00 UTC 2022
Hyperglycemia induces miR-26-5p down-regulation to overexpress PFKFB3 and accelerate epithelial-mesenchymal transition in gastric cancer.
Bioengineered
He X,Cheng X,Ding J,Xiong M,Chen B,Cao G
Tue Feb 01 00:00:00 UTC 2022
Discovery and characterization of heterogeneous and multipotent fibroblast populations isolated from excised cleft lip tissue.
Stem cell research & therapy
Parisi L,Rihs S,La Scala GC,Schnyder I,Katsaros C,Degen M
Thu Sep 08 00:00:00 UTC 2022
ERK1/2 Signalling Pathway Regulates Tubulin-Binding Cofactor B Expression and Affects Astrocyte Process Formation after Acute Foetal Alcohol Exposure.
Brain sciences
Zheng Y,Huo J,Yang M,Zhang G,Wan S,Chen X,Zhang B,Liu H
Wed Jun 22 00:00:00 UTC 2022
Chemical profile and miscarriage prevention evaluation of Jiao-Ai Decoction, a classical traditional Chinese formula.
Journal of pharmaceutical and biomedical analysis
Zhang M,Geng T,Jing SH,Li L,Wang K,Zhang Q,Zhang Y,Zhou SK,Gao P,Ding AW,Zhang L
Fri Aug 05 00:00:00 UTC 2022
Orientin downregulating oxidative stress-mediated endoplasmic reticulum stress and mitochondrial dysfunction through AMPK/SIRT1 pathway in rat nucleus pulposus cells in vitro and attenuated intervertebral disc degeneration in vivo.
Apoptosis : an international journal on programmed cell death
Zhang Z,Wu J,Teng C,Wang J,Yu J,Jin C,Wang L,Wu L,Lin Z,Yu Z,Lin Z
Thu Dec 01 00:00:00 UTC 2022
The role of regulatory T cells on the activation of astrocytes in the brain of high-fat diet mice following lead exposure.
Chemico-biological interactions
Wu L,Li S,Li C,He B,Lv L,Wang J,Wang J,Wang W,Zhang Y
Wed Jan 05 00:00:00 UTC 2022
A Recombinant Genotype I Japanese Encephalitis Virus Expressing a Gaussia Luciferase Gene for Antiviral Drug Screening Assay and Neutralizing Antibodies Detection.
International journal of molecular sciences
Li C,Chen X,Hu J,Jiang D,Cai D,Li Y
Thu Dec 08 00:00:00 UTC 2022
Developing a Mathematical Model of Intracellular Calcium Dynamics for Evaluating Combined Anticancer Effects of Afatinib and RP4010 in Esophageal Cancer.
International journal of molecular sciences
Chang Y,Funk M,Roy S,Stephenson E,Choi S,Kojouharov HV,Chen B,Pan Z
Thu Feb 03 00:00:00 UTC 2022
HepaCAM‑PIK3CA axis regulates the reprogramming of glutamine metabolism to inhibit prostate cancer cell proliferation.
International journal of oncology
He Z,Gao Y,Li T,Yu C,Ou L,Luo C
Fri Apr 01 00:00:00 UTC 2022
Antitumor and Antibacterial Efficacy of Gallium Nanoparticles Coated by Ellagic Acid.
Dose-response : a publication of International Hormesis Society
El-Sonbaty SM,Moawed FS,Kandil EI,M Tamamm A
Sat Feb 19 00:00:00 UTC 2022
Acetyl-11-Keto-Beta Boswellic Acid (AKBA) Protects Lens Epithelial Cells Against H2O2-Induced Oxidative Injury and Attenuates Cataract Progression by Activating Keap1/Nrf2/HO-1 Signaling.
Frontiers in pharmacology
Yang T,Lin X,Li H,Zhou X,Fan F,Yang J,Luo Y,Liu X
Fri Jul 29 00:00:00 UTC 2022
Venetoclax enhances DNA damage induced by XPO1 inhibitors: A novel mechanism underlying the synergistic antileukaemic effect in acute myeloid leukaemia.
Journal of cellular and molecular medicine
Yu H,Wu S,Liu S,Li X,Gai Y,Lin H,Wang Y,Edwards H,Ge Y,Wang G
Sun May 01 00:00:00 UTC 2022
Cystine Induced-mTORC2 Activation through Promoting Sin1 Phosphorylation to Suppress Cancer Cell Ferroptosis.
Molecular nutrition & food research
Wang G,Chen L,Qin S,Zheng Y,Xia C,Yao J,Wang P,Deng L
Thu Dec 01 00:00:00 UTC 2022
Functional analyses of two interferon-stimulated gene 15 (ISG15) copies in large yellow croaker, Larimichthys crocea.
Fish & shellfish immunology
Shen B,Zhang S,Li F,Xu J,Zhang Y,Zhang J
Tue Nov 01 00:00:00 UTC 2022
Effect of condylar chondrocyte exosomes on condylar cartilage osteogenesis in rats under tensile stress.
Frontiers in bioengineering and biotechnology
Shi Y,Shao J,Zhang Z,Zhang J,Lu H
Sat Dec 24 00:00:00 UTC 2022
KDM4C Contributes to Trophoblast-like Stem Cell Conversion from Porcine-Induced Pluripotent Stem Cells (piPSCs) via Regulating CDX2.
International journal of molecular sciences
Yu S,Shen Q,Zhang R,Wu X,Zhang J,Zhao W,Wu X,Li N,Peng S,Zhang S,Yang F,Hua J
Fri Jul 08 00:00:00 UTC 2022
CDGSH iron sulfur domain 2 mitigates apoptosis, oxidative stress and inflammatory response caused by oxygen-glucose deprivation/reoxygenation in HT22 hippocampal neurons by Akt-Nrf2-activated pathway.
Metabolic brain disease
Ren X,Yu J,Guo L,Zhang Z
Sat Oct 01 00:00:00 UTC 2022
Downregulation of FXYD2 Is Associated with Poor Prognosis and Increased Regulatory T Cell Infiltration in Clear Cell Renal Cell Carcinoma.
Journal of immunology research
Zhang Z,Tang Y,Li L,Yang W,Xu Y,Zhou J,Ma K,Zhang K,Zhuang H,Gong Y,Gong K
Tue Nov 01 00:00:00 UTC 2022
Berberine prevents lethal EV71 neurological infection in newborn mice.
Frontiers in pharmacology
Cui G,Wang H,Yang C,Zhou X,Wang J,Wang T,Ma T
Fri Nov 18 00:00:00 UTC 2022
Evacetrapib Elicits Antitumor Effects on Colorectal Cancer by Inhibiting the Wnt/β-Catenin Signaling Pathway and Activating the JNK Signaling Pathway.
Biological & pharmaceutical bulletin
Hu L,Dong H,He L,Shi M,Xiang N,Su Y,Wang C,Tian Y,Hu Y,Wang H,Liu H,Wen C,Yang X
Thu Sep 08 00:00:00 UTC 2022
Disulfiram alleviates pristane-induced lupus via inhibiting GSDMD-mediated pyroptosis.
Cell death discovery
Zhuang L,Luo X,Wu S,Lin Z,Zhang Y,Zhai Z,Yang F,Li Y,Zhuang J,Luo G,Xu W,He Y,Sun E
Sat Sep 03 00:00:00 UTC 2022
The synergistic anticancer effect of the bromodomain inhibitor OTX015 and histone deacetylase 6 inhibitor WT-161 in osteosarcoma.
Cancer cell international
Yu B,Liu L,Cai F,Peng Y,Tang X,Zeng D,Li T,Zhang F,Liang Y,Yuan X,Li J,Dai Z,Liao Q,Lv XB
Tue Feb 08 00:00:00 UTC 2022
CD137 Signal Mediates Cardiac Ischemia-Reperfusion Injury by Regulating the Necrosis of Cardiomyocytes.
Journal of cardiovascular translational research
Xu Y,Chen R,Yan J,Zang G,Shao C,Wang Z
Sat Oct 01 00:00:00 UTC 2022
BMP6 Promotes the Secretion of 17 Beta-Estradiol and Progesterone in Goat Ovarian Granulosa Cells.
Animals : an open access journal from MDPI
Song S,Ding W,Yao H,Wang L,Li B,Wang Y,Tang X,Zhang Y,Huang D,Xu D,Zhao Z
Fri Aug 19 00:00:00 UTC 2022
Basolateral Amygdala Reactive Microglia May Contribute to Synaptic Impairment and Depressive-Like Behavior in Mice with Bone Cancer Pain.
Neurochemical research
Jiang L,Hao J,Yang XL,Zhu JX,Wang Y,Huang YL,Sun YE,Mao YT,Ni K,Gu XP,Ma ZL
Tue Nov 01 00:00:00 UTC 2022
Tnfaip2 promotes atherogenesis by enhancing oxidative stress induced inflammation.
Molecular immunology
Jin G,Liu Y,Xu W,Li Y,Zhang H,Qiu S,Gao C,Liu S
Tue Nov 01 00:00:00 UTC 2022
Ferroptosis promotes microtubule-associated protein tau aggregation via GSK-3β activation and proteasome inhibition.
Molecular neurobiology
Wang S,Jiang Y,Liu Y,Liu Q,Sun H,Mei M,Liao X
Tue Mar 01 00:00:00 UTC 2022
The combination of decitabine and aspirin inhibits tumor growth and metastasis in non-small cell lung cancer.
The Journal of international medical research
Xu M,Song B,Yang X,Li N
Fri Jul 01 00:00:00 UTC 2022
Transmissible Endoplasmic Reticulum Stress Mediated by Extracellular Vesicles from Adipocyte Promoting the Senescence of Adipose-Derived Mesenchymal Stem Cells in Hypertrophic Obesity.
Oxidative medicine and cellular longevity
Fang J,Li L,Cao X,Yue H,Fu W,Chen Y,Xu Z,Zhao Q,Zhao J,Wang Y,Liang W
Tue Aug 30 00:00:00 UTC 2022
Mitochondrial and glucose metabolic dysfunctions in granulosa cells induce impaired oocytes of polycystic ovary syndrome through Sirtuin 3.
Free radical biology & medicine
Zhang Q,Ren J,Wang F,Pan M,Cui L,Li M,Qu F
Fri Jul 01 00:00:00 UTC 2022
High glucose and palmitic acid induces neuronal senescence by NRSF/REST elevation and the subsequent mTOR-related autophagy suppression.
Molecular brain
Xue WJ,He CF,Zhou RY,Xu XD,Xiang LX,Wang JT,Wang XR,Zhou HG,Guo JC
Mon Jul 18 00:00:00 UTC 2022
MFG-E8 alleviates intervertebral disc degeneration by suppressing pyroptosis and extracellular matrix degradation in nucleus pulposus cells via Nrf2/TXNIP/NLRP3 axis.
Cell death discovery
Ma H,Xie C,Chen Z,He G,Dai Z,Cai H,Zhang H,Lu H,Wu H,Hu X,Zhou K,Zheng G,Xu H,Xu C
Tue Apr 19 00:00:00 UTC 2022
Hydrogen-generating Si-based agent protects against skin flap ischemia-reperfusion injury in rats.
Scientific reports
Otani N,Tomita K,Kobayashi Y,Kuroda K,Koyama Y,Kobayashi H,Kubo T
Wed Apr 13 00:00:00 UTC 2022
Isoliquiritigenin-mediated miR-23a-3p inhibition activates PGC-1α to alleviate alcoholic liver injury.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang L,Kong L,Xu S,Wang X,Huang K,Wang S,Wu J,Wang C,Sun H,Liu K,Meng Q
Tue Feb 01 00:00:00 UTC 2022
miR-186-5p Dysregulation in Serum Exosomes from Patients with AMI Aggravates Atherosclerosis via Targeting LOX-1.
International journal of nanomedicine
Ding J,Li H,Liu W,Wang X,Feng Y,Guan H,Chen Z
Thu Dec 29 00:00:00 UTC 2022
Comprehensive Analysis of the Potential Immune-Related Biomarker ATG101 that Regulates Apoptosis of Cholangiocarcinoma Cells After Photodynamic Therapy.
Frontiers in pharmacology
Zhang ZJ,Wang KP,Huang YP,Jin C,Jiang H,Xiong L,Chen ZY,Wen Y,Liu ZT,Mo JG
Sat Jul 16 00:00:00 UTC 2022
Ginkgo biloba leaf extract mitigates cisplatin-induced chronic renal interstitial fibrosis by inhibiting the epithelial-mesenchymal transition of renal tubular epithelial cells mediated by the Smad3/TGF-β1 and Smad3/p38 MAPK pathways.
Chinese medicine
Wei C,Zhang Y,Zhong X,Lu S,Zou X,Yang Y,Huang S,Huang Z
Mon Feb 21 00:00:00 UTC 2022
Optineurin-mediated mitophagy as a potential therapeutic target for intervertebral disc degeneration.
Frontiers in pharmacology
Hu Z,Wang Y,Gao X,Zhang Y,Liu C,Zhai Y,Chang X,Li H,Li Y,Lou J,Li C
Sat Sep 17 00:00:00 UTC 2022
Alleviating effect of quercetin on cadmium-induced oxidative damage and apoptosis by activating the Nrf2-keap1 pathway in BRL-3A cells.
Frontiers in pharmacology
Wang J,Wang K,Ding L,Zhao P,Zhang C,Wang H,Yang Z,Liu Z
Tue Aug 30 00:00:00 UTC 2022
Transglutaminase 3 Attenuates Skin Inflammation in Psoriasis by Inhibiting NF-κB Activation through Phosphorylated STAT3-TET3 Signaling.
The Journal of investigative dermatology
Ling S,Xu B,Luo Y,Fang X,Liu X,Wang A,Zhou Y,Zhang S,Zong W,Li W,Yao X
Tue Nov 01 00:00:00 UTC 2022
Systematic characterization of the metabolites of defatted walnut powder extract in vivo and screening of the mechanisms against NAFLD by UPLC-Q-Exactive Orbitrap MS combined with network pharmacology.
Journal of ethnopharmacology
Ren SM,Zhang QZ,Jiang M,Chen ML,Xu XJ,Wang DM,Pan YN,Liu XQ
Tue Mar 01 00:00:00 UTC 2022
Preliminary evaluation of a small interfering RNA molecular probe targeting murine double minute 2 in breast cancer.
Nuclear medicine communications
Wang X,Xu P,Jiao Y,Luan S,Gao Y,Zhao C,Fu P
Mon Aug 01 00:00:00 UTC 2022
GRP75 Modulates Endoplasmic Reticulum-Mitochondria Coupling and Accelerates Ca2+-Dependent Endothelial Cell Apoptosis in Diabetic Retinopathy.
Biomolecules
Li Y,Li HY,Shao J,Zhu L,Xie TH,Cai J,Wang W,Cai MX,Wang ZL,Yao Y,Wei TT
Tue Nov 29 00:00:00 UTC 2022
SPATA33 affects the formation of cell adhesion complex by interacting with CTNNA3 in TM4 cells.
Cell and tissue research
Zhang Y
Fri Jul 01 00:00:00 UTC 2022
Loureirin C ameliorates ischemia and reperfusion injury in rats by inhibiting the activation of the TLR4/NF-κB pathway and promoting TLR4 degradation.
Phytotherapy research : PTR
Xu J,Liu J,Li Q,Mi Y,Zhou D,Wang J,Chen G,Liang D,Li N,Hou Y
Thu Dec 01 00:00:00 UTC 2022
Ginsenoside CK induces apoptosis in triple-negative breast cancer cells by targeting glutamine metabolism.
Biochemical pharmacology
Zhang B,Fu R,Duan Z,Shen S,Zhu C,Fan D
Mon Aug 01 00:00:00 UTC 2022
Protective role of small extracellular vesicles derived from HUVECs treated with AGEs in diabetic vascular calcification.
Journal of nanobiotechnology
Guo B,Shan SK,Xu F,Lin X,Li FX,Wang Y,Xu QS,Zheng MH,Lei LM,Li CC,Zhou ZA,Ullah MHE,Wu F,Liao XB,Yuan LQ
Sat Jul 16 00:00:00 UTC 2022
Epigenetic silencing of JAM3 promotes esophageal cancer development by activating Wnt signaling.
Clinical epigenetics
Yang W,Guo C,Herman JG,Zhu C,Lv H,Su X,Zhang L,Zhang M,Guo M
Fri Dec 02 00:00:00 UTC 2022
Novel bioactive hybrid Celecoxib-HDAC Inhibitor, induces apoptosis in human acute lymphoblastic leukemia cells.
Bioorganic & medicinal chemistry
Liu J,Zhang L,Guo L,Zeng Y,Guo Q,Yang C,Shu J,Liu W,Yang L
Thu Dec 01 00:00:00 UTC 2022
Isolation, characterization, and functional study of extracellular vesicles derived from Leishmania tarentolae.
Frontiers in cellular and infection microbiology
Shokouhy M,Sarvnaz H,Taslimi Y,Lajevardi MS,Habibzadeh S,Mizbani A,Shekari F,Behbahani M,Torrecilhas AC,Rafati S
Tue Aug 23 00:00:00 UTC 2022
Rhodiola activates macrophage migration inhibitory factor to alleviate non-alcoholic fatty liver disease.
Life sciences
Liu J,Li D,Dun Y,Li H,Ripley-Gonzalez JW,Zhang J,Qiu L,You B,Liu S
Tue Nov 01 00:00:00 UTC 2022
Intake of flavonoids from Astragalus membranaceus ameliorated brain impairment in diabetic mice via modulating brain-gut axis.
Chinese medicine
Li X,Zhao T,Gu J,Wang Z,Lin J,Wang R,Duan T,Li Z,Dong R,Wang W,Hong KF,Liu Z,Huang W,Gui D,Zhou H,Xu Y
Sat Feb 12 00:00:00 UTC 2022
7-Methoxyisoflavone suppresses vascular endothelial inflammation by inhibiting the expression of endothelial adhesion molecules.
European journal of pharmacology
Zhu X,Peng J,Yang L,Guo Y,Wang P,Liu K,Zhu J,Deng S,Liang M
Sat Oct 15 00:00:00 UTC 2022
Neonatal gene therapy achieves sustained disease rescue of maple syrup urine disease in mice.
Nature communications
Pontoizeau C,Simon-Sola M,Gaborit C,Nguyen V,Rotaru I,Tual N,Colella P,Girard M,Biferi MG,Arnoux JB,Rötig A,Ottolenghi C,de Lonlay P,Mingozzi F,Cavazzana M,Schiff M
Tue Jun 07 00:00:00 UTC 2022
Ejection of damaged mitochondria and their removal by macrophages ensure efficient thermogenesis in brown adipose tissue.
Cell metabolism
Rosina M,Ceci V,Turchi R,Chuan L,Borcherding N,Sciarretta F,Sánchez-Díaz M,Tortolici F,Karlinsey K,Chiurchiù V,Fuoco C,Giwa R,Field RL,Audano M,Arena S,Palma A,Riccio F,Shamsi F,Renzone G,Verri M,Crescenzi A,Rizza S,Faienza F,Filomeni G,Kooijman S,Rufini S,de Vries AAF,Scaloni A,Mitro N,Tseng YH,Hidalgo A,Zhou B,Brestoff JR,Aquilano K,Lettieri-Barbato D
Tue Apr 05 00:00:00 UTC 2022
Macamide B Pretreatment Attenuates Neonatal Hypoxic-Ischemic Brain Damage of Mice Induced Apoptosis and Regulates Autophagy via the PI3K/AKT Signaling Pathway.
Molecular neurobiology
Yang X,Wang M,Zhou Q,Bai Y,Liu J,Yang J,Li L,Li G,Luo L
Sun May 01 00:00:00 UTC 2022
Angiotensin II type-2 receptor activation in alveolar macrophages mediates protection against cigarette smoke-induced chronic obstructive pulmonary disease.
Pharmacological research
Mei D,Liao W,Gan PXL,Tran QTN,Chan CCMY,Heng CKM,Wong WSF
Sat Oct 01 00:00:00 UTC 2022
Identification of oncogenes and tumor-suppressor genes with hepatocellular carcinoma: A comprehensive analysis based on TCGA and GEO datasets.
Frontiers in genetics
Zhu Y,Wang Y,Hu M,Lu X,Sun G
Thu Feb 02 00:00:00 UTC 2023
San-Huang-Yi-Shen Capsule Ameliorates Diabetic Kidney Disease through Inducing PINK1/Parkin-Mediated Mitophagy and Inhibiting the Activation of NLRP3 Signaling Pathway.
Journal of diabetes research
Li H,Wang Y,Su X,Wang Q,Zhang S,Sun W,Zhang T,Dong M,Zhang Z,Lv S
Mon Nov 28 00:00:00 UTC 2022
Novel Antiviral Activity of Ethyl 3-Hydroxyhexanoate Against Coxsackievirus B Infection.
Frontiers in microbiology
Olasunkanmi OI,Mageto J,Avala Ntsigouaye J,Yi M,Fei Y,Chen Y,Chen S,Xu W,Lin L,Zhao W,Wang Y,Zhong ZH
Tue May 03 00:00:00 UTC 2022
Parkin-mediated mitophagy protects against TNF-α-induced stress in bone marrow mesenchymal stem cells.
Experimental gerontology
Fan P,Yu XY,Chen CH,Gao JW,Xu YZ,Xie XH,Wang YT
Fri Jul 01 00:00:00 UTC 2022
Circular RNA circBNC2 inhibits epithelial cell G2-M arrest to prevent fibrotic maladaptive repair.
Nature communications
Wang P,Huang Z,Peng Y,Li H,Lin T,Zhao Y,Hu Z,Zhou Z,Zhou W,Liu Y,Hou FF
Mon Oct 31 00:00:00 UTC 2022
Chicoric acid attenuates tumor necrosis factor-α-induced inflammation and apoptosis via the Nrf2/HO-1, PI3K/AKT and NF-κB signaling pathways in C28/I2 cells and ameliorates the progression of osteoarthritis in a rat model.
International immunopharmacology
Qu Y,Shen Y,Teng L,Huang Y,Yang Y,Jian X,Fan S,Wu P,Fu Q
Sat Oct 01 00:00:00 UTC 2022
Muscone and (+)-Borneol Cooperatively Strengthen CREB Induction of Claudin 5 in IL-1β-Induced Endothelium Injury.
Antioxidants (Basel, Switzerland)
Li YC,Li Y,Zhang YN,Zhao Q,Zhang PL,Sun MR,Liu BL,Yang H,Li P
Tue Jul 26 00:00:00 UTC 2022
Identification of a novel native peptide derived from 60S ribosomal protein L23a that translationally regulates p53 to reduce myocardial ischemia-reperfusion.
Pharmacological research
Yin A,Feng M,Zhang L,Cheng Z,Li Y,Qian L
Sat Jan 01 00:00:00 UTC 2022
Identification of cytochrome c oxidase subunit 4 isoform 1 as a positive regulator of influenza virus replication.
Frontiers in microbiology
He J,Huang H,Li B,Li H,Zhao Y,Li Y,Ye W,Qi W,Tang W,Wang L
Sat Aug 06 00:00:00 UTC 2022
TOP2A deficiency leads to human recurrent spontaneous abortion and growth retardation of mouse pre-implantation embryos.
Molecular medicine (Cambridge, Mass.)
Duan Y,Fu H,Huang J,Yin N,Liu L,Liu X
Fri Dec 30 00:00:00 UTC 2022
Chemerin Regulates the Proliferation and Migration of Pulmonary Arterial Smooth Muscle Cells via the ERK1/2 Signaling Pathway.
Frontiers in pharmacology
Peng L,Chen Y,Li Y,Feng P,Zheng Y,Dong Y,Yang Y,Wang R,Li A,Yan J,Shang F,Tang P,Chen D,Gao Y,Huang W
Tue Apr 05 00:00:00 UTC 2022
Gestational bisphenol A exposure impairs hepatic lipid metabolism by altering mTOR/CRTC2/SREBP1 in male rat offspring.
Human & experimental toxicology
Yang Q,Mao Y,Wang J,Yu H,Zhang X,Pei X,Duan Z,Xiao C,Ma M
Mon Sep 26 00:00:00 UTC 2022
Hydrogen sulfide alleviates apoptosis and autophagy induced by beryllium sulfate in 16HBE cells.
Journal of applied toxicology : JAT
Liu YP,Yuan XY,Li XY,Wang Y,Sun ZB,Deng WH,Lei YD,Huang L,Jiang TY,Zhang ZH
Tue Feb 01 00:00:00 UTC 2022
Pectin supplement alleviates gut injury potentially through improving gut microbiota community in piglets.
Frontiers in microbiology
Dang G,Wang W,Zhong R,Wu W,Chen L,Zhang H
Tue Jan 03 00:00:00 UTC 2023
Prunella vulgaris can improve the pregnancy outcomes of experimental autoimmune thyroiditis rats by inhibiting Th1/Th17 immune responses.
Journal of reproductive immunology
Zhu Q,Muyayalo KP,Xu QH,Wang J,Wang H,Liao AH
Tue Feb 01 00:00:00 UTC 2022
Indole-3-propionic acid alleviates ischemic brain injury in a mouse middle cerebral artery occlusion model.
Experimental neurology
Xie Y,Zou X,Han J,Zhang Z,Feng Z,Ouyang Q,Hua S,Liu Z,Li C,Cai Y,Zou Y,Tang Y,Jiang X
Fri Jul 01 00:00:00 UTC 2022
Circadian Rhythm Disorders Aggravate Periodontitis by Modulating BMAL1.
International journal of molecular sciences
Liu X,Cao N,Liu X,Deng Y,Xin Y,Fu R,Xin X,Hou Y,Yu W
Mon Dec 26 00:00:00 UTC 2022
Effects of iron oxide nanoparticles as T2-MRI contrast agents on reproductive system in male mice.
Journal of nanobiotechnology
Yang H,Wang H,Wen C,Bai S,Wei P,Xu B,Xu Y,Liang C,Zhang Y,Zhang G,Wen H,Zhang L
Wed Mar 02 00:00:00 UTC 2022
Moxibustion alleviates decreased ovarian reserve in rats by restoring the PI3K/AKT signaling pathway.
Journal of integrative medicine
Li HX,Shi L,Liang SJ,Fang CC,Xu QQ,Lu G,Wang Q,Cheng J,Shen J,Shen MH
Tue Mar 01 00:00:00 UTC 2022
STING-mediated inflammation contributes to Gao binge ethanol feeding model.
Journal of cellular physiology
Wang L,You HM,Meng HW,Pan XY,Chen X,Bi YH,Zhang YF,Li JJ,Yin NN,Zhang ZW,Huang C,Li J
Tue Feb 01 00:00:00 UTC 2022
Trans-Anethole Alleviates Subclinical Necro-Haemorrhagic Enteritis-Induced Intestinal Barrier Dysfunction and Intestinal Inflammation in Broilers.
Frontiers in microbiology
Yu C,Wang D,Tong Y,Li Q,Yang W,Wang T,Yang Z
Fri Apr 08 00:00:00 UTC 2022
Puerarin-V prevents the progression of hypoxia- and monocrotaline-induced pulmonary hypertension in rodent models.
Acta pharmacologica Sinica
Chen D,Zhang HF,Yuan TY,Sun SC,Wang RR,Wang SB,Fang LH,Lyu Y,Du GH
Thu Sep 01 00:00:00 UTC 2022
Let-7i-5p promotes a malignant phenotype in nasopharyngeal carcinoma via inhibiting tumor-suppressive autophagy.
Cancer letters
You B,Zhang P,Gu M,Yin H,Fan Y,Yao H,Pan S,Xie H,Cheng T,Liu H,You Y,Liu J
Sun Apr 10 00:00:00 UTC 2022
CEND1 deficiency induces mitochondrial dysfunction and cognitive impairment in Alzheimer's disease.
Cell death and differentiation
Xie W,Guo D,Li J,Yue L,Kang Q,Chen G,Zhou T,Wang H,Zhuang K,Leng L,Li H,Chen Z,Gao W,Zhang J
Thu Dec 01 00:00:00 UTC 2022
FSH inhibits autophagy and lysosomal biogenesis to regulate protein degradation in cultured goat Sertoli cells.
Molecular and cellular endocrinology
Xi H,Ren F,Li Y,Xian M,Wang L,Hu J
Sat Jan 15 00:00:00 UTC 2022
SRA Suppresses Antiviral Innate Immune Response in Macrophages by Limiting TBK1 K63 Ubiquitination via Deubiquitinase USP15.
Microbiology spectrum
Li L,Luo J,Zhu Z,Xu Q,Wang P,Chang B,Wang D,Yu L,Lu X,Zhou J,Zuo D,Chen Q
Wed Dec 21 00:00:00 UTC 2022
Aucubin promotes bone-fracture healing via the dual effects of anti-oxidative damage and enhancing osteoblastogenesis of hBM-MSCs.
Stem cell research & therapy
Wang K,Zhou C,Li L,Dai C,Wang Z,Zhang W,Xu J,Zhu Y,Pan Z
Fri Aug 19 00:00:00 UTC 2022
Tumor-associated macrophages promote migration and invasion via modulating IL-6/STAT3 signaling in renal cell carcinoma.
International immunopharmacology
Chen S,Qian S,Zhang L,Pan X,Qu F,Yu Y,Gu Z,Cui X,Shen H
Sat Oct 01 00:00:00 UTC 2022
Evodiamine suppresses the progression of non-small cell lung carcinoma via endoplasmic reticulum stress-mediated apoptosis pathway in vivo and in vitro.
International journal of immunopathology and pharmacology
Li Y,Wang Y,Wang X,Jin L,Yang L,Zhu J,Wang H,Zheng F,Cui H,Li X,Jia Y
Fri Apr 08 00:00:00 UTC 2022
MicroRNA-200c Affects Milk Fat Synthesis by Targeting PANK3 in Ovine Mammary Epithelial Cells.
International journal of molecular sciences
Hao Z,Wang J,Luo Y,Hu J,Liu X,Li S,Li M,Shi B,Hu L,Liu Y,Zhen H
Fri Dec 09 00:00:00 UTC 2022
Glia Maturation Factor β as a Novel Independent Prognostic Biomarker and Potential Therapeutic Target of Kidney Renal Clear Cell Carcinoma.
Frontiers in oncology
Zhu T,Wang T,Feng Z,Gao F,Zhang J,Jin C,Tian H,Xu J,Chen H,Ou Q,Wang J,Xu G,Lu L
Fri Jul 22 00:00:00 UTC 2022
Inhibition of MAT2A suppresses osteoclastogenesis and prevents ovariectomy-induced bone loss.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kang H,Guo Q,Dong Y,Peng R,Song K,Wang J,Liu H,Zhu M,Zhao H,Guan H,Li F
Tue Feb 01 00:00:00 UTC 2022
MiR-328-5p inhibits the adipogenic differentiation of hMSCs by targeting fatty acid synthase.
Folia histochemica et cytobiologica
Li X,He S,Wu P,Zhou Y,Long K,Wang T
Mon Jan 02 00:00:00 UTC 2023
125I seeds inhibit proliferation and promote apoptosis in cholangiocarcinoma cells by regulating the AGR2-mediated p38 MAPK pathway.
Cancer letters
Zhou X,Zhang W,Dou M,Li Z,Liu Z,Li J,Tian C,Yao Y,Wang C,Li Y,Chen P,Han X,Jiao D
Sat Jan 01 00:00:00 UTC 2022
BMAL1 regulates Propionibacterium acnes-induced skin inflammation via REV-ERBα in mice.
International journal of biological sciences
Li F,Lin L,He Y,Sun G,Dong D,Wu B
Thu Apr 14 00:00:00 UTC 2022
Calpain-1 mediates vascular remodelling and fibrosis via HIF-1α in hypoxia-induced pulmonary hypertension.
Journal of cellular and molecular medicine
Deng H,Tian X,Sun H,Liu H,Lu M,Wang H
Sun May 01 00:00:00 UTC 2022
Didymin Suppresses Microglia Pyroptosis and Neuroinflammation Through the Asc/Caspase-1/GSDMD Pathway Following Experimental Intracerebral Hemorrhage.
Frontiers in immunology
Gu L,Sun M,Li R,Zhang X,Tao Y,Yuan Y,Luo X,Xie Z
Mon Mar 14 00:00:00 UTC 2022
Arsenic exposure via drinking water during pregnancy and lactation induces autism-like behaviors in male offspring mice.
Chemosphere
Zhang X,Mei D,Li Y,You M,Wang D,Yao D,Xu Y,Zhai L,Wang Y
Tue Mar 01 00:00:00 UTC 2022
Nrf2 Overexpression Decreases Vincristine Chemotherapy Sensitivity Through the PI3K-AKT Pathway in Adult B-Cell Acute Lymphoblastic Leukemia.
Frontiers in oncology
Wang L,Liu X,Kang Q,Pan C,Zhang T,Feng C,Chen L,Wei S,Wang J
Sat Jul 16 00:00:00 UTC 2022
P2X7R/NLRP3 signaling pathway-mediated pyroptosis and neuroinflammation contributed to cognitive impairment in a mouse model of migraine.
The journal of headache and pain
Wang Y,Shan Z,Zhang L,Fan S,Zhou Y,Hu L,Wang Y,Li W,Xiao Z
Sat Jul 02 00:00:00 UTC 2022
Altered Liver Metabolism, Mitochondrial Function, Oxidative Status, and Inflammatory Response in Intrauterine Growth Restriction Piglets with Different Growth Patterns before Weaning.
Metabolites
Wang J,Zhu P,Zheng X,Ma Z,Cui C,Wu C,Zeng X,Guan W,Chen F
Tue Nov 01 00:00:00 UTC 2022
Integrated Bioinformatic Analysis Reveals TXNRD1 as a Novel Biomarker and Potential Therapeutic Target in Idiopathic Pulmonary Arterial Hypertension.
Frontiers in medicine
Lin W,Tang Y,Zhang M,Liang B,Wang M,Zha L,Yu Z
Mon Jan 02 00:00:00 UTC 2023
Lipid-lowering activity of metformin-soluble soybean polysaccharide nanoparticles.
Food & function
He H,Liu M,He R,Zhao W
Mon Oct 03 00:00:00 UTC 2022
MiR-103a-3p Promotes Zika Virus Replication by Targeting OTU Deubiquitinase 4 to Activate p38 Mitogen-Activated Protein Kinase Signaling Pathway.
Frontiers in microbiology
Ye H,Kang L,Yan X,Li S,Huang Y,Mu R,Duan X,Chen L
Thu Mar 24 00:00:00 UTC 2022
Multifunctional photodynamic/photothermal nano-agents for the treatment of oral leukoplakia.
Journal of nanobiotechnology
Lin L,Song C,Wei Z,Zou H,Han S,Cao Z,Zhang X,Zhang G,Ran J,Cai Y,Han W
Fri Mar 04 00:00:00 UTC 2022
DNA damage-induced translocation of mitochondrial factor HIGD1A into the nucleus regulates homologous recombination and radio/chemo-sensitivity.
Oncogene
Chen B,Xu F,Gao Y,Hu G,Zhu K,Lu H,Xu A,Chen S,Wu L,Zhao G
Tue Mar 01 00:00:00 UTC 2022
MYB proto-oncogene like 2 promotes hepatocellular carcinoma growth and glycolysis via binding to the Optic atrophy 3 promoter and activating its expression.
Bioengineered
Liu M,Du Q,Mao G,Dai N,Zhang F
Tue Mar 01 00:00:00 UTC 2022
Human Amnion Epithelial Cells and Their Derived Exosomes Alleviate Sepsis-Associated Acute Kidney Injury via Mitigating Endothelial Dysfunction.
Frontiers in medicine
Chi D,Chen Y,Xiang C,Yao W,Wang H,Zheng X,Xu D,Li N,Xie M,Wang S,Liu G,Li S,Yang L
Wed Apr 13 00:00:00 UTC 2022
The lncRNA BDNF-AS/WDR5/FBXW7 axis mediates ferroptosis in gastric cancer peritoneal metastasis by regulating VDAC3 ubiquitination.
International journal of biological sciences
Huang G,Xiang Z,Wu H,He Q,Dou R,Lin Z,Yang C,Huang S,Song J,Di Z,Wang S,Xiong B
Fri Apr 15 00:00:00 UTC 2022
Inhibition of glutamine transporter ASCT2 mitigates bleomycin-induced pulmonary fibrosis in mice.
Acta histochemica
Lian N,Jin H,Zhu W,Zhang C,Qi Y,Jiang M,Mao J,Lu X,Zhao F,Xi B,Qi X,Li Y
Thu Dec 01 00:00:00 UTC 2022
MCPIP1 alleviates inflammatory response through inducing autophagy in Aspergillus fumigatus keratitis.
International immunopharmacology
Han F,Shen L,Ma H,Wang L,Guo H,Wu X
Thu Dec 01 00:00:00 UTC 2022
ProNGF/NGF Modulates Autophagy and Apoptosis through PI3K/Akt/mTOR and ERK Signaling Pathways following Cerebral Ischemia-Reperfusion in Rats.
Oxidative medicine and cellular longevity
Li Y,Wu F,Zhou M,Zhou J,Cui S,Guo J,Wu J,He L
Mon Apr 11 00:00:00 UTC 2022
Oxaloacetate acid ameliorates paraquat-induced acute lung injury by alleviating oxidative stress and mitochondrial dysfunction.
Frontiers in pharmacology
Li W,Li M,Chen K,Tang Y,Yin R,Lan L,Hong G
Wed Nov 02 00:00:00 UTC 2022
Vitamin D Deficiency Impacts Exposure and Response of Pravastatin in Male Rats by Altering Hepatic OATPs.
Frontiers in pharmacology
Peng J,Yang G,Huang Z
Tue Mar 08 00:00:00 UTC 2022
MicroRNA-20a promotes non-small cell lung cancer proliferation by upregulating PD-L1 by targeting PTEN.
Oncology letters
Gong J,Shen Y,Jiang F,Wang Y,Chu L,Sun J,Shen P,Chen M
Sun May 01 00:00:00 UTC 2022
Inhibition of Bruton's Tyrosine Kinase Alleviates Monocrotaline-Induced Pulmonary Arterial Hypertension by Modulating Macrophage Polarization.
Oxidative medicine and cellular longevity
Yu M,Wu X,Peng L,Yang M,Zhou H,Xu J,Wang J,Wang H,Xie W,Kong H
Fri Sep 09 00:00:00 UTC 2022
Olanzapine-induced lipid disturbances: A potential mechanism through the gut microbiota-brain axis.
Frontiers in pharmacology
Zhu Z,Gu Y,Zeng C,Yang M,Yu H,Chen H,Zhang B,Cai H
Tue Aug 23 00:00:00 UTC 2022
Salidroside Ameliorates Radiation Damage by Reducing Mitochondrial Oxidative Stress in the Submandibular Gland.
Antioxidants (Basel, Switzerland)
Sun YM,Wang XY,Zhou XR,Zhang C,Liu KJ,Zhang FY,Xiang B
Thu Jul 21 00:00:00 UTC 2022
Ketogenesis acts as an endogenous protective programme to restrain inflammatory macrophage activation during acute pancreatitis.
EBioMedicine
Zhang L,Shi J,Du D,Niu N,Liu S,Yang X,Lu P,Shen X,Shi N,Yao L,Zhang R,Hu G,Lu G,Zhu Q,Zeng T,Liu T,Xia Q,Huang W,Xue J
Fri Apr 01 00:00:00 UTC 2022
Loureirin A Exerts Antikeloid Activity by Antagonizing the TGF-β1/Smad Signalling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ma H,Duan X,Zhang R,Li H,Guo Y,Tian Y,Huang M,Chen G,Wang Z,Li L
Thu Dec 21 00:00:00 UTC 2023
Diet-Induced High Serum Levels of Trimethylamine-N-oxide Enhance the Cellular Inflammatory Response without Exacerbating Acute Intracerebral Hemorrhage Injury in Mice.
Oxidative medicine and cellular longevity
Li C,Zhu L,Dai Y,Zhang Z,Huang L,Wang TJ,Fu P,Li Y,Wang J,Jiang C
Thu Mar 24 00:00:00 UTC 2022
Development and Validation of Novel Prognostic Models for Immune-Related Genes in Osteosarcoma.
Frontiers in molecular biosciences
Li J,Su L,Xiao X,Wu F,Du G,Guo X,Kong F,Yao J,Zhu H
Sat Jul 16 00:00:00 UTC 2022
Hypoxic mesenchymal stem cell-derived extracellular vesicles ameliorate renal fibrosis after ischemia-reperfusion injure by restoring CPT1A mediated fatty acid oxidation.
Stem cell research & therapy
Gao Z,Zhang C,Peng F,Chen Q,Zhao Y,Chen L,Wang X,Chen X
Sat May 07 00:00:00 UTC 2022
A plant-derived natural photosynthetic system for improving cell anabolism.
Nature
Chen P,Liu X,Gu C,Zhong P,Song N,Li M,Dai Z,Fang X,Liu Z,Zhang J,Tang R,Fan S,Lin X
Thu Dec 01 00:00:00 UTC 2022
Niban apoptosis regulator 1 promotes gemcitabine resistance by activating the focal adhesion kinase signaling pathway in bladder cancer.
Journal of Cancer
Tong S,Yin H,Fu J,Li Y
Tue Mar 15 00:00:00 UTC 2022
Dysregulation of SIRT3 SUMOylation Confers AML Chemoresistance via Controlling HES1-Dependent Fatty Acid Oxidation.
International journal of molecular sciences
Zhang Y,Shen Y,Wei W,Wang W,Jiang D,Ren Y,Peng Z,Fan Q,Cheng J,Ma J
Wed Jul 27 00:00:00 UTC 2022
Combinatorial approaches for mitigating resistance to KRAS-targeted therapies.
The Biochemical journal
Warren HR,Ross SJ,Smith PD,Coulson JM,Prior IA
Fri Sep 30 00:00:00 UTC 2022
Effects of STAT3 Inhibitor BP-1-102 on The Proliferation, Invasiveness, Apoptosis and Neurosphere Formation of Glioma Cells in Vitro.
Cell biochemistry and biophysics
Zhang CC,Wu T,Guan L,Wang YJ,Yao RQ,Gao DS,Li F
Thu Dec 01 00:00:00 UTC 2022
Targeting glutamine metabolism in hepatic stellate cells alleviates liver fibrosis.
Cell death & disease
Yin X,Peng J,Gu L,Liu Y,Li X,Wu J,Xu B,Zhuge Y,Zhang F
Mon Nov 14 00:00:00 UTC 2022
An electrochemical nitric oxide generator for in-home inhalation therapy in pulmonary artery hypertension.
BMC medicine
Liu Y,Zhu Y,Jiang C,Su Z,Yan Y,Feng B,Mao W,Zhang Y,Wang X,Xu Z,Zhang H
Thu Dec 15 00:00:00 UTC 2022
Artesunate Inhibits the Cell Growth in Colorectal Cancer by Promoting ROS-Dependent Cell Senescence and Autophagy.
Cells
Huang Z,Gan S,Zhuang X,Chen Y,Lu L,Wang Y,Qi X,Feng Q,Huang Q,Du B,Zhang R,Liu Z
Tue Aug 09 00:00:00 UTC 2022
Longterm Increased S100B Enhances Hippocampal Progenitor Cell Proliferation in a Transgenic Mouse Model.
International journal of molecular sciences
Rodrigues L,Wartchow KM,Buchfelder M,Souza DO,Gonçalves CA,Kleindienst A
Wed Aug 24 00:00:00 UTC 2022
Mettl3 regulates hypertrophic differentiation of chondrocytes through modulating Dmp1 mRNA via Ythdf1-mediated m6A modification.
Bone
He Y,Wang W,Luo P,Wang Y,He Z,Dong W,Jia M,Yu X,Yang B,Wang J
Tue Nov 01 00:00:00 UTC 2022
Magnesium Hydride Ameliorates Endotoxin-Induced Acute Respiratory Distress Syndrome by Inhibiting Inflammation, Oxidative Stress, and Cell Apoptosis.
Oxidative medicine and cellular longevity
Shi X,Zhu L,Wang S,Zhu W,Li Q,Wei J,Feng D,Liu M,Chen Y,Sun X,Lu H,Lv X
Tue May 10 00:00:00 UTC 2022
Long Non-coding RNA ANRIL Downregulation Alleviates Neuroinflammation in an Ischemia Stroke Model via Modulation of the miR-671-5p/NF-κB Pathway.
Neurochemical research
Deng L,Jiang J,Chen S,Lin X,Zuo T,Hu Q,Wu Y,Fan X,Dong Z
Fri Jul 01 00:00:00 UTC 2022
The Role and Mechanism of CREBH Regulating SIRT3 in Metabolic Associated Fatty Liver Disease.
Life sciences
Junli Z,Shuhan W,Yajuan Z,Xiaoling D,Jiahuan L,Keshu X
Sat Oct 01 00:00:00 UTC 2022
Curcumin regulates anti-inflammatory responses by AXL/JAK2/STAT3 signaling pathway in experimental autoimmune encephalomyelitis.
Neuroscience letters
Sun M,Liu N,Sun J,Li X,Wang H,Zhang W,Xie Q,Wang M
Wed Sep 14 00:00:00 UTC 2022
Corrigendum to "Berberine Reduces Renal Cell Pyroptosis in Golden Hamsters with Diabetic Nephropathy through the Nrf2-NLRP3-Caspase-1-GSDMD Pathway".
Evidence-based complementary and alternative medicine : eCAM
Ding B,Geng S,Hou X,Ma X,Xu H,Yang F,Liu K,Liang W,Ma G
Thu Aug 18 00:00:00 UTC 2022
Resveratrol inhibits the progression of premature senescence partially by regulating v-rel avian reticuloendotheliosis viral oncogene homolog A (RELA) and sirtuin 1 (SIRT1).
Renal failure
He S,Zhou M,Zheng H,Wang Y,Wu S,Gao Y,Chen J
Thu Dec 01 00:00:00 UTC 2022
PM2.5-Induced Programmed Myocardial Cell Death via mPTP Opening Results in Deteriorated Cardiac Function in HFpEF Mice.
Cardiovascular toxicology
Wu T,Tong M,Chu A,Wu K,Niu X,Zhang Z
Mon Aug 01 00:00:00 UTC 2022
Ellagic Acid Alleviates Mice Intestinal Ischemia-Reperfusion Injury: A Study Based on Transcriptomics Combined with Functional Experiments.
Chemistry & biodiversity
Fan S,Xu Y,Li K,Li B,Diao Y
Tue Nov 01 00:00:00 UTC 2022
Pu-erh tea increases the metabolite Cinnabarinic acid to improve circadian rhythm disorder-induced obesity.
Food chemistry
Hu S,Hu C,Luo L,Zhang H,Zhao S,Liu Z,Zeng L
Tue Nov 15 00:00:00 UTC 2022
Roles of C/EBP-homologous protein and histone H3 lysine 4 methylation in arsenic-induced mitochondrial apoptosis in hepatocytes.
Toxicology and industrial health
Liang C,Xie RJ,Wang JL,Zhang YW,Zhang JY,Yang Q,Han B
Tue Nov 01 00:00:00 UTC 2022
Ergosterol Peroxide Inhibits Porcine Epidemic Diarrhea Virus Infection in Vero Cells by Suppressing ROS Generation and p53 Activation.
Viruses
Liu Y,Wang X,Wang J,Zhang J,Duan C,Wang J
Tue Feb 15 00:00:00 UTC 2022
Loss of BAF (mSWI/SNF) chromatin-remodeling ATPase Brg1 causes multiple malformations of cortical development in mice.
Human molecular genetics
Jin Y,Gao X,Lu M,Chen G,Yang X,Ren N,Song Y,Hou C,Li J,Liu Q,Gao J
Mon Oct 10 00:00:00 UTC 2022
NLRP3 Inflammasome Activation Enhances ADK Expression to Accelerate Epilepsy in Mice.
Neurochemical research
Zhang H,Yu S,Xia L,Peng X,Wang S,Yao B
Tue Mar 01 00:00:00 UTC 2022
NF-κB and AP-1 are required for the lipopolysaccharide-induced expression of MCP-1, CXCL1, and Cx43 in cultured rat dorsal spinal cord astrocytes.
Frontiers in molecular neuroscience
Lu Y,Li B,Xu A,Liang X,Xu T,Jin H,Xie Y,Wang R,Liu X,Gao X,Han Y,Zeng J
Tue Aug 16 00:00:00 UTC 2022
Astragali Radix-Coptis Rhizoma Herb Pair Attenuates Atherosclerosis in ApoE-/- Mice by Regulating the M1/M2 and Th1/Th2 Immune Balance and Activating the STAT6 Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Li Z,Ma D,Wang Y,Wu S,Wang L,Jiang Y,Zhang Y,Li X
Sun May 01 00:00:00 UTC 2022
mTORC1 Mediates the Processes of Lysine Regulating Satellite Cells Proliferation, Apoptosis, and Autophagy.
Metabolites
Liu M,Yue Z,Zhang B,Li F,Liu L,Li F
Thu Aug 25 00:00:00 UTC 2022
Structural Variability of Lipoarabinomannan Modulates Innate Immune Responses within Infected Alveolar Epithelial Cells.
Cells
Liu H,Gui X,Chen S,Fu W,Li X,Xiao T,Hou J,Jiang T
Fri Jan 21 00:00:00 UTC 2022
P53 maintains gallid alpha herpesvirus 1 replication by direct regulation of nucleotide metabolism and ATP synthesis through its target genes.
Frontiers in microbiology
Xu L,Chen Z,Zhang Y,Cui L,Liu Z,Li X,Liu S,Li H
Tue Dec 20 00:00:00 UTC 2022
The expression of m6A enzymes in the hippocampus of diabetic cognitive impairment mice and the possible improvement of YTHDF1.
Brain research
Li M,Zhong X,Zhao Z,Zeng Z,Yuan Q,Xiao X,Zu X,Liu J
Tue Feb 15 00:00:00 UTC 2022
Jujuboside A ameliorates high fat diet and streptozotocin induced diabetic nephropathy via suppressing oxidative stress, apoptosis, and enhancing autophagy.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Zhong Y,Luo R,Liu Q,Zhu J,Lei M,Liang X,Wang X,Peng X
Sat Jan 01 00:00:00 UTC 2022
MicroRNA-122a aggravates intestinal ischemia/reperfusion injury by promoting pyroptosis via targeting EGFR-NLRP3 signaling pathway.
Life sciences
Wang F,Gu L,Wang Y,Sun D,Zhao Y,Meng Q,Yin L,Xu L,Lu X,Peng J,Lin Y,Sun P
Sat Oct 15 00:00:00 UTC 2022
Duck Tembusu Virus Inhibits Type I Interferon Production through the JOSD1-SOCS1-IRF7 Negative-Feedback Regulation Pathway.
Journal of virology
Huang S,Huang J,Cui M,Wu X,Wang M,Zhu D,Chen S,Liu M,Zhao X,Wu Y,Yang Q,Zhang S,Ou X,Mao S,Gao Q,Yu Y,Tian B,Liu Y,Zhang L,Yin Z,Jing B,Chen X,Cheng A,Jia R
Wed Sep 28 00:00:00 UTC 2022
Genome-wide association studies identify miRNA-194 as a prognostic biomarker for gastrointestinal cancer by targeting ATP6V1F, PPP1R14B, BTF3L4 and SLC7A5.
Frontiers in oncology
Huang P,Xia L,Guo Q,Huang C,Wang Z,Huang Y,Qin S,Leng W,Li D
Wed Jan 11 00:00:00 UTC 2023
Overexpression of NCAPG in ovarian cancer is associated with ovarian cancer proliferation and apoptosis via p38 MAPK signaling pathway.
Journal of ovarian research
Yu H,Zou D,Ni N,Zhang S,Zhang Q,Yang L
Fri Aug 19 00:00:00 UTC 2022
Enhancement of anti-inflammatory effect of cattle bile by fermentation and its inhibition of neuroinflammation on microglia by inhibiting NLRP3 inflammasome.
Journal of bioscience and bioengineering
Pei Y,Cheng F,Li W,Yu Q,Ma C,Zou Y,Xu T,Liu S,Zhang S,Wang Q
Tue Feb 01 00:00:00 UTC 2022
Probing the Potential Mechanism of Quercetin and Kaempferol against Heat Stress-Induced Sertoli Cell Injury: Through Integrating Network Pharmacology and Experimental Validation.
International journal of molecular sciences
Liu DL,Liu SJ,Hu SQ,Chen YC,Guo J
Thu Sep 22 00:00:00 UTC 2022
Kaempferol From Penthorum chinense Pursh Attenuates Hepatic Ischemia/Reperfusion Injury by Suppressing Oxidative Stress and Inflammation Through Activation of the Nrf2/HO-1 Signaling Pathway.
Frontiers in pharmacology
Chen Y,Li T,Tan P,Shi H,Cheng Y,Cai T,Bai J,Du Y,Fu W
Sat Jul 16 00:00:00 UTC 2022
Lgr5 + cell fate regulation by coordination of metabolic nuclear receptors during liver repair.
Theranostics
Qin D,Liu S,Lu Y,Yan Y,Zhang J,Cao S,Chen M,Chen N,Huang W,Wang L,Chen X,Zhang L
Thu Sep 29 00:00:00 UTC 2022
KIAA1199 Correlates With Tumor Microenvironment and Immune Infiltration in Lung Adenocarcinoma as a Potential Prognostic Biomarker.
Pathology oncology research : POR
Shen X,Mo X,Tan W,Mo X,Li L,Yu F,He J,Deng Z,Xing S,Chen Z,Yang J
Thu Dec 01 00:00:00 UTC 2022
Dl-3-n-Butylphthalide Improves Neuroinflammation in Mice with Repeated Cerebral Ischemia-Reperfusion Injury through the Nrf2-Mediated Antioxidant Response and TLR4/MyD88/NF-κB Signaling Pathway.
Oxidative medicine and cellular longevity
Gao Y,Hu M,Niu X,Li M,Xu L,Xiao Y,Zhang J,Wang H,Li L,Chu B,Lv P
Tue Dec 13 00:00:00 UTC 2022
Clostridium butyricum inhibits the progression of colorectal cancer and alleviates intestinal inflammation via the myeloid differentiation factor 88 (MyD88)-nuclear factor-kappa B (NF-κB) signaling pathway.
Annals of translational medicine
Zhou M,Yuan W,Yang B,Pei W,Ma J,Feng Q
Fri Apr 01 00:00:00 UTC 2022
Rhein Inhibits the Progression of Chemoresistant Lung Cancer Cell Lines via the Stat3/Snail/MMP2/MMP9 Pathway.
BioMed research international
Liu J,Ding D,Liu F,Chen Y
Thu Mar 31 00:00:00 UTC 2022
Detection of netrin-1 as a novel biomarker for diagnosis and chemotherapeutic monitoring of lung cancer.
The Journal of international medical research
Zhao Y,Song J,Ding X,Hao Y,Cao L
Wed Jun 01 00:00:00 UTC 2022
Effects of the probiotic, Lactobacillus rhamnosus GG, on ulcer pathogenesis, HSP70 stress protein and nitric oxide levels in stress induced ulcer.
Biotechnic & histochemistry : official publication of the Biological Stain Commission
Isik M,Ozbayer C,Donmez DB,Colak E,Ustuner MC,Erol K,Degirmenci I
Mon Aug 01 00:00:00 UTC 2022
Systematic analyses identify the anti-fibrotic role of lncRNA TP53TG1 in IPF.
Cell death & disease
Sun J,Guo Y,Chen T,Jin T,Ma L,Ai L,Guo J,Niu Z,Yang R,Wang Q,Yu X,Gao H,Zhang Y,Su W,Song X,Ji W,Zhang Q,Huang M,Fan X,Du Z,Liang H
Sat Jun 04 00:00:00 UTC 2022
ADAR1p110 promotes Enterovirus D68 replication through its deaminase domain and inhibition of PKR pathway.
Virology journal
Zhang K,Wang S,Chen T,Tu Z,Huang X,Zang G,Wu C,Fan X,Liu J,Tian Y,Cheng Y,Lu N,Zhang G
Thu Dec 22 00:00:00 UTC 2022
A novel synthetic chalcone derivative, 2,4,6-trimethoxy-4'-nitrochalcone (Ch-19), exerted anti-tumor effects through stimulating ROS accumulation and inducing apoptosis in esophageal cancer cells.
Cell stress & chaperones
Yang Y,Wu H,Zou X,Chen Y,He R,Jin Y,Zhou B,Ge C,Yang Y
Tue Nov 01 00:00:00 UTC 2022
Protective Effect of Flavonoids against Methylglyoxal-Induced Oxidative Stress in PC-12 Neuroblastoma Cells and Its Structure-Activity Relationships.
Molecules (Basel, Switzerland)
Zhang D,Li X,He X,Xing Y,Jiang B,Xiu Z,Bao Y,Dong Y
Sat Nov 12 00:00:00 UTC 2022
HDAC6 Inhibition Alleviates Anesthesia and Surgery-Induced Less Medial Prefrontal-Dorsal Hippocampus Connectivity and Cognitive Impairment in Aged Rats.
Molecular neurobiology
Chen J,Liu S,Wang X,Huang J,Phillips J,Ma D,Ouyang W,Tong J
Sat Oct 01 00:00:00 UTC 2022
LncRNA WEE2-AS1 Knockdown Inhibits the Proliferation, Migration and 3 Invasion of Glioma Cells via Regulating miR-29b-2-5p/TPM3 Axis.
Oncology research
Jia Z,Qian Z,Tang Y,Li X,Shi Y,Xin H,Fan Y,Wu H
Thu Apr 07 00:00:00 UTC 2022
The Photoperiod Regulates Granulosa Cell Apoptosis through the FSH-Nodal/ALK7 Signaling Pathway in Phodopus sungorus.
Animals : an open access journal from MDPI
Qi Y,Xue HL,Zheng YJ,Yin YF,Xu WL,Xu JH,Wu M,Chen L,Xu LX
Fri Dec 16 00:00:00 UTC 2022
The Effect and Mechanism of Duodenal-Jejunal Bypass to Treat Type 2 Diabetes Mellitus in a Rat Model.
Obesity facts
Chen X,Zhang Q,Yang Q,Huang Z,Liao G,Wang Z
Thu May 19 00:00:00 UTC 2022
Adipose-derived stem cells alleviate liver injury induced by type 1 diabetes mellitus by inhibiting mitochondrial stress and attenuating inflammation.
Stem cell research & therapy
Hou Y,Ding W,Wu P,Liu C,Ding L,Liu J,Wang X
Fri Apr 01 00:00:00 UTC 2022
FUNDC2 promotes liver tumorigenesis by inhibiting MFN1-mediated mitochondrial fusion.
Nature communications
Li S,Han S,Zhang Q,Zhu Y,Zhang H,Wang J,Zhao Y,Zhao J,Su L,Li L,Zhou D,Ye C,Feng XH,Liang T,Zhao B
Fri Jun 17 00:00:00 UTC 2022
Overexpression of sFlt-1 represses ox-LDL-induced injury of HUVECs by activating autophagy via PI3K/AKT/mTOR pathway.
Microvascular research
Zhou YH,Tang YZ,Guo LY,Zheng LL,Zhang D,Yang CY,Wang W
Sat Jan 01 00:00:00 UTC 2022
Impact of Selective Renal Afferent Denervation on Oxidative Stress and Vascular Remodeling in Spontaneously Hypertensive Rats.
Antioxidants (Basel, Switzerland)
Wu LL,Zhang Y,Li XZ,Du XL,Gao Y,Wang JX,Wang XL,Chen Q,Li YH,Zhu GQ,Tan X
Fri May 20 00:00:00 UTC 2022
Adrenomedullin is an Important Pathological Mediator in Progression of Chronic Neuropathic Pain.
Frontiers in bioscience (Landmark edition)
Wang C,Xue Y,Lu Q,Shi Y,Tang W,Wang D
Wed Jul 13 00:00:00 UTC 2022
Pt(II)-NHC Complex Induces ROS-ERS-Related DAMP Balance to Harness Immunogenic Cell Death in Hepatocellular Carcinoma.
Journal of medicinal chemistry
Bian M,Fan R,Yang Z,Chen Y,Xu Z,Lu Y,Liu W
Thu Feb 10 00:00:00 UTC 2022
Gomisin A enhances the antitumor effect of paclitaxel by suppressing oxidative stress in ovarian cancer.
Oncology reports
Wang T,Liu J,Huang X,Zhang C,Shangguan M,Chen J,Wu S,Chen M,Yang Z,Zhao S
Tue Nov 01 00:00:00 UTC 2022
NEDD4 attenuates phosgene-induced acute lung injury through the inhibition of Notch1 activation.
Journal of cellular and molecular medicine
Shao Y,Jiang Z,He D,Shen J
Sun May 01 00:00:00 UTC 2022
A mouse-adapted CVA6 strain exhibits neurotropism and triggers systemic manifestations in a novel murine model.
Emerging microbes & infections
Li D,Sun T,Tao L,Ji W,Zhu P,Liang R,Zhang Y,Chen S,Yang H,Jin Y,Duan G
Thu Dec 01 00:00:00 UTC 2022
12/15-Lipoxygenase Regulation of Diabetic Cognitive Dysfunction Is Determined by Interfering with Inflammation and Cell Apoptosis.
International journal of molecular sciences
Chen Q,Zheng Q,Yang Y,Luo Y,Wang H,Li H,Yang L,Hu C,Zhang J,Li Y,Xia H,Chen Z,Ma J,Tian X,Yang J
Fri Aug 12 00:00:00 UTC 2022
Tetrandrine overcomes drug resistance mediated by bone marrow microenvironment by regulating the expression of P-glycoprotein in acute leukemia.
Hematology (Amsterdam, Netherlands)
Zhou X,Jin N,Chen B
Thu Dec 01 00:00:00 UTC 2022
Costunolide protects against alcohol-induced liver injury by regulating gut microbiota, oxidative stress and attenuating inflammation in vivo and in vitro.
Phytotherapy research : PTR
Mao J,Zhan H,Meng F,Wang G,Huang D,Liao Z,Chen M
Tue Mar 01 00:00:00 UTC 2022
Parthenolide modulates cerebral ischemia-induced microglial polarization and alleviates neuroinflammatory injury via the RhoA/ROCK pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhang Y,Miao L,Peng Q,Fan X,Song W,Yang B,Zhang P,Liu G,Liu J
Sat Oct 01 00:00:00 UTC 2022
The human FOXM1 homolog promotes basal progenitor cell proliferation and cortical folding in mouse.
EMBO reports
Wang W,Su L,Ji F,Zhang D,Wang Y,Zhao J,Jiao RD,Zhang M,Huang E,Jiang H,Zhang J,Jiao J
Thu Feb 03 00:00:00 UTC 2022
Thermosensitive Hydrogel Incorporating Prussian Blue Nanoparticles Promotes Diabetic Wound Healing via ROS Scavenging and Mitochondrial Function Restoration.
ACS applied materials & interfaces
Xu Z,Liu Y,Ma R,Chen J,Qiu J,Du S,Li C,Wu Z,Yang X,Chen Z,Chen T
Wed Mar 30 00:00:00 UTC 2022
Proteomic and functional analyses of the periodic membrane skeleton in neurons.
Nature communications
Zhou R,Han B,Nowak R,Lu Y,Heller E,Xia C,Chishti AH,Fowler VM,Zhuang X
Thu Jun 09 00:00:00 UTC 2022
Stratifin as a novel diagnostic biomarker in serum for diffuse alveolar damage.
Nature communications
Arakawa N,Ushiki A,Abe M,Matsuyama S,Saito Y,Kashiwada T,Horimasu Y,Gemma A,Tatsumi K,Hattori N,Tsushima K,Miyashita K,Saito K,Nakamura R,Toyoda T,Ogawa K,Sato M,Takamatsu K,Mori K,Nishiya T,Izumi T,Ohno Y,Saito Y,Hanaoka M
Tue Oct 04 00:00:00 UTC 2022
Effects of Low-Intensity Pulsed Ultrasound on the Migration and Homing of Human Amnion-Derived Mesenchymal Stem Cells to Ovaries in Rats With Premature Ovarian Insufficiency.
Cell transplantation
Ling L,Hou J,Wang Y,Shu H,Huang Y
Wed Oct 26 00:00:00 UTC 2022
Chitosan Oligosaccharide Attenuates Lipopolysaccharide-Induced Intestinal Barrier Dysfunction through Suppressing the Inflammatory Response and Oxidative Stress in Mice.
Antioxidants (Basel, Switzerland)
Tao W,Wang G,Pei X,Sun W,Wang M
Sun Jul 17 00:00:00 UTC 2022
PERK Regulates the Sensitivity of Hepatocellular Carcinoma Cells to High-LET Carbon Ions via either Apoptosis or Ferroptosis.
Journal of Cancer
Zheng X,Liu B,Liu X,Li P,Zhang P,Ye F,Zhao T,Kuang Y,Chen W,Jin X,Li Q
Tue Jan 25 00:00:00 UTC 2022
The Effective Analysis for Blue Honeysuckle Extract in the Treatment of Hepatocellular Carcinoma.
Evidence-based complementary and alternative medicine : eCAM
Zhang CP,Li WH,Liu JR,Li GD,Zhang HP,Wei JF,Chen HS,Zhao JL,Wang YF,Lv Q,Liu M
Tue Oct 11 00:00:00 UTC 2022
Baicalein suppresses high glucose-induced inflammation and apoptosis in trophoblasts by targeting the miRNA-17-5p-Mfn1/2-NF-κB pathway.
Placenta
Li J,Wang Y,Wu T,Li S,Sun YN,Liu ZH
Fri Apr 01 00:00:00 UTC 2022
Isovitexin protects against acute liver injury by targeting PTEN, PI3K and BiP via modification of m6A.
European journal of pharmacology
Huang Y,Chen S,Pang L,Feng Z,Su H,Zhu W,Wei J
Tue Feb 15 00:00:00 UTC 2022
Interleukin-6-derived cancer-associated fibroblasts activate STAT3 pathway contributing to gemcitabine resistance in cholangiocarcinoma.
Frontiers in pharmacology
Kittirat Y,Suksawat M,Thongchot S,Padthaisong S,Phetcharaburanin J,Wangwiwatsin A,Klanrit P,Sangkhamanon S,Titapun A,Loilome W,Saya H,Namwat N
Tue Sep 13 00:00:00 UTC 2022
EPLINβ Is Involved in the Assembly of Cadherin-catenin Complexes in Osteoblasts and Affects Bone Formation.
Acta histochemica et cytochemica
Miyazaki S,Funamoto T,Sekimoto T,Kurogi S,Ohta T,Nagai T,Tajima T,Imasaka M,Yoshinobu K,Araki K,Araki M,Choijookhuu N,Hishikawa Y,Chosa E
Wed Jun 29 00:00:00 UTC 2022
S-palmitoylation of PCSK9 induces sorafenib resistance in liver cancer by activating the PI3K/AKT pathway.
Cell reports
Sun Y,Zhang H,Meng J,Guo F,Ren D,Wu H,Jin X
Tue Aug 16 00:00:00 UTC 2022
Molecular and functional characterization of teleost-specific Interleukin-17N in yellow catfish (Pelteobagrus fulvidraco).
International journal of biological macromolecules
Zhou X,Jiang XX,Zhang GR,Ji W,Shi ZC,Ma XF,Wei KJ
Tue Nov 01 00:00:00 UTC 2022
Gasdermin D is involved in switching from apoptosis to pyroptosis in TLR4-mediated renal tubular epithelial cells injury in diabetic kidney disease.
Archives of biochemistry and biophysics
Yuan S,Wang Y,Li Z,Chen X,Song P,Chen A,Qu Z,Wen S,Liu H,Zhu X
Fri Sep 30 00:00:00 UTC 2022
Optimized mobilization of MHC class I- and II- restricted immunity by dendritic cell vaccine potentiates cancer therapy.
Theranostics
Shi Y,Liu Y,Huang J,Luo Z,Guo X,Jiang M,Li X,Lu Y,Liu X,Shan X,Luo L,You J
Tue May 17 00:00:00 UTC 2022
Candida albicans induces upregulation of programmed death ligand 1 in oral squamous cell carcinoma.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Wang X,Zhao W,Zhang W,Wu S,Yan Z
Sun May 01 00:00:00 UTC 2022
Placental expression of estrogen-related receptor gamma is reduced in fetal growth restriction pregnancies and is mediated by hypoxia†.
Biology of reproduction
Zou Z,Harris LK,Forbes K,Heazell AEP
Mon Sep 12 00:00:00 UTC 2022
Mitigation Effects of Selenium Nanoparticles on Depression-Like Behavior Induced by Fluoride in Mice via the JAK2-STAT3 Pathway.
ACS applied materials & interfaces
Yang J,Li H,Hao Z,Jing X,Zhao Y,Cheng X,Ma H,Wang J,Wang J
Wed Jan 26 00:00:00 UTC 2022
Proteomic and Biochemical Approaches Elucidate the Role of Millimeter-Wave Irradiation in Wheat Growth under Flooding Stress.
International journal of molecular sciences
Komatsu S,Tsutsui Y,Furuya T,Yamaguchi H,Hitachi K,Tsuchida K,Tani M
Thu Sep 08 00:00:00 UTC 2022
Cefotetan-bound human RKIP involves in Ras/Raf1/MEK/ERK signaling pathway.
Acta biochimica et biophysica Sinica
Guo C,Xu H,Zhou Y,Wu Z,Jiang B,Chen H,Lin D
Sun Dec 25 00:00:00 UTC 2022
Downregulation of miR-30b-5p Facilitates Chondrocyte Hypertrophy and Apoptosis via Targeting Runx2 in Steroid-Induced Osteonecrosis of the Femoral Head.
International journal of molecular sciences
Lin L,Yu Y,Liu K,Jiang Y,Zhou Z
Sat Sep 24 00:00:00 UTC 2022
Protective effect of active components of Eucommia ulmoides leaves on gastric ulcers in rats: Involvement of the PI3K/Akt/NF-κB pathway.
Journal of food science
Gong M,Li Q,Guo H,Cui B,Liu Y,Wang P,Zhu H,Liu X,Dai L,Wang Z
Fri Jul 01 00:00:00 UTC 2022
Non-invasive photobiomodulation treatment in an Alzheimer Disease-like transgenic rat model.
Theranostics
Yang L,Wu C,Parker E,Li Y,Dong Y,Tucker L,Brann DW,Lin HW,Zhang Q
Thu Apr 21 00:00:00 UTC 2022
ATRX-dependent SVCT2 mediates macrophage infiltration in the glioblastoma xenograft model.
Journal of neurophysiology
Shang J,Wang Y,Li Z,Jiang L,Bai Q,Zhang X,Xiao G,Zhang J
Sun May 01 00:00:00 UTC 2022
Tubuloside B, isolated from Cistanche tubulosa, a promising agent against M1 macrophage activation via synergistically targeting Mob1 and ERK1/2.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Xiao L,Yao J,Miao Y,Ou B,Wang J,Huang Y,Zhou B,Ge L,Tian J,Zeng X
Thu Sep 01 00:00:00 UTC 2022
Transplantation of Human Umbilical Cord Mesenchymal Stem Cells-Derived Neural Stem Cells Pretreated with Neuregulin1β Ameliorate Cerebral Ischemic Reperfusion Injury in Rats.
Biomolecules
Zhai QY,Ren YQ,Ni QS,Song ZH,Ge KL,Guo YL
Thu Mar 10 00:00:00 UTC 2022
Oxidative stress activates Ryr2-Ca2+ and apoptosis to promote PM2.5-induced heart injury of hyperlipidemia mice.
Ecotoxicology and environmental safety
Meng M,Jia R,Wei M,Meng X,Zhang X,Du R,Sun W,Wang L,Song L
Tue Mar 01 00:00:00 UTC 2022
Hypoxia-inducible Factor Regulates Ten-eleven Translocated Methylcytosine Dioxygenase 1-c-Myc Binding Involved in Depression-like Behavior in Prenatal Hypoxia Offspring.
Neuroscience
Zeng H,Wei B,Liu J,Lu L,Li L,Wang B,Sun M
Sat Oct 15 00:00:00 UTC 2022
Poloxamer 407 and Hyaluronic Acid Thermosensitive Hydrogel-Encapsulated Ginsenoside Rg3 to Promote Skin Wound Healing.
Frontiers in bioengineering and biotechnology
Peng X,Ding C,Zhao Y,Hao M,Liu W,Yang M,Xiao F,Zheng Y
Sat Jul 23 00:00:00 UTC 2022
Salidroside inhibited cerebral ischemia/reperfusion-induced oxidative stress and apoptosis via Nrf2/Trx1 signaling pathway.
Metabolic brain disease
Li F,Mao Q,Wang J,Zhang X,Lv X,Wu B,Yan T,Jia Y
Thu Dec 01 00:00:00 UTC 2022
Puffball spores improve wound healing in a diabetic rat model.
Frontiers in endocrinology
He T,Sun P,Liu B,Wan S,Fang P,Chen J,Huang G,Min W
Tue Sep 20 00:00:00 UTC 2022
Cinnamic Acid Improved Lipopolysaccharide-Induced Depressive-Like Behaviors by Inhibiting Neuroinflammation and Oxidative Stress in Mice.
Pharmacology
Zhuo R,Cheng X,Luo L,Yang L,Zhao Y,Zhou Y,Peng L,Jin X,Cui L,Liu F,Yang L
Wed May 04 00:00:00 UTC 2022
Mesenchymal-to-epithelial transition of osteoblasts induced by Fam20c knockout.
Genes & genomics
Geng YW,Zhang Z,Jin H,Da JL,Zhang K,Wang JQ,Guo YY,Zhang B,Li Y
Tue Feb 01 00:00:00 UTC 2022
Systematic pan-cancer analysis identifies RALA as a tumor targeting immune therapeutic and prognostic marker.
Frontiers in immunology
Jin H,Qin S,He J,Xiao J,Li Q,Mao Y,Zhao L
Tue Dec 06 00:00:00 UTC 2022
Venetoclax enhances NK cell killing sensitivity of AML cells through the NKG2D/NKG2DL activation pathway.
International immunopharmacology
Wu HY,Li KX,Pan WY,Guo MQ,Qiu DZ,He YJ,Li YH,Huang YX
Tue Mar 01 00:00:00 UTC 2022
Circular RNA circCCDC85A inhibits breast cancer progression via acting as a miR-550a-5p sponge to enhance MOB1A expression.
Breast cancer research : BCR
Meng L,Chang S,Sang Y,Ding P,Wang L,Nan X,Xu R,Liu F,Gu L,Zheng Y,Li Z,Sang M
Tue Jan 04 00:00:00 UTC 2022
CircFGGY Inhibits Cell Growth, Invasion and Epithelial-Mesenchymal Transition of Hepatocellular Carcinoma via Regulating the miR-545-3p/Smad7 Axis.
Frontiers in cell and developmental biology
Feng KL,Diao N,Zhou ZW,Fang CK,Wang JN,Zhang Y,Luo R,Zhong C
Sat Jul 16 00:00:00 UTC 2022
Deficiency of a novel lncRNA-HRAT protects against myocardial ischemia reperfusion injury by targeting miR-370-3p/RNF41 pathway.
Frontiers in cardiovascular medicine
Zheng X,Zhong T,Yu F,Duan J,Tang Y,Liu Y,Li M,Sun D,Yin D
Fri Sep 30 00:00:00 UTC 2022
A positive feedback circuit comprising p21 and HIF-1α aggravates hypoxia-induced radioresistance of glioblastoma by promoting Glut1/LDHA-mediated glycolysis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Jin X,Kuang Y,Li L,Li H,Zhao T,He Y,Di C,Kang J,Yuan L,Yu B,Li Q
Tue Mar 01 00:00:00 UTC 2022
Identification of key genes and pathways revealing the central regulatory mechanism of brain-derived glucagon-like peptide-1 on obesity using bioinformatics analysis.
Frontiers in neuroscience
Shao Y,Tian J,Yang Y,Hu Y,Zhu Y,Shu Q
Tue Aug 23 00:00:00 UTC 2022
Tumor microenvironment-related gene selenium-binding protein 1 (SELENBP1) is associated with immunotherapy efficacy and survival in colorectal cancer.
BMC gastroenterology
Zhu C,Wang S,Du Y,Dai Y,Huai Q,Li X,Du Y,Dai H,Yuan W,Yin S,Wang H
Mon Oct 17 00:00:00 UTC 2022
Jumonji-C domain-containing protein 5 suppresses proliferation and aerobic glycolysis in pancreatic cancer cells in a c-Myc-dependent manner.
Cellular signalling
Wang H,Wang J,Liu J,Wang Y,Xia G,Huang X
Sun May 01 00:00:00 UTC 2022
Glibenclamide Directly Prevents Neuroinflammation by Targeting SUR1-TRPM4-Mediated NLRP3 Inflammasome Activation In Microglia.
Molecular neurobiology
He Y,Chang Y,Peng Y,Zhu J,Liu K,Chen J,Wu Y,Ji Z,Lin Z,Wang S,Gupta S,Zang N,Pan S,Huang K
Sat Oct 01 00:00:00 UTC 2022
Host-induced gene silencing of PcCesA3 and PcOSBP1 confers resistance to Phytophthora capsici in Nicotiana benthamiana through NbDCL3 and NbDCL4 processed small interfering RNAs.
International journal of biological macromolecules
Wang Z,Gao X,Zhong S,Li Y,Shi M,Zhang B,Zhang S,Shen H,Liu X
Thu Dec 01 00:00:00 UTC 2022
GPR12 Inhibits Apoptosis in Epithelial Ovarian Cancer via the Activation of ERK1/2 Signaling.
Frontiers in oncology
Wang L,Yang D,Zhang Y,Jiao Y
Sun Jul 31 00:00:00 UTC 2022
PIN1 protects auditory hair cells from senescence via autophagy.
PeerJ
Lv Z,Zhang Y,Cao H,Liu Q,Feng X,Yin H,Wang B
Fri Feb 10 00:00:00 UTC 2023
Role of the cGAS-STING Pathway in Aging-related Endothelial Dysfunction.
Aging and disease
Yu H,Liao K,Hu Y,Lv D,Luo M,Liu Q,Huang L,Luo S
Thu Dec 01 00:00:00 UTC 2022
The oral microbiome in young women at different stages of periodontitis: Prevotella dominant in stage III periodontitis.
Frontiers in cellular and infection microbiology
Zhao Y,Feng Y,Ye Q,Hu J,Feng Y,Ouyang Z,Zhao J,Chen Y,Tan L,Chen N,Dusenge MA,Su X,Guo Y
Tue Dec 20 00:00:00 UTC 2022
Nanosilica-Anchored Polycaprolactone/Chitosan Nanofibrous Bioscaffold to Boost Osteogenesis for Bone Tissue Engineering.
Molecules (Basel, Switzerland)
Ge S,Zhu X,Zhang C,Jia D,Shang W,Ding C,Yang J,Feng Y
Tue Dec 13 00:00:00 UTC 2022
A Novel High-Throughput Screening Method for a Human Multicentric Osteosarcoma-Specific Antibody and Biomarker Using a Phage Display-Derived Monoclonal Antibody.
Cancers
Hayashi T,Yamamoto N,Kurosawa G,Tajima K,Kondo M,Hiramatsu N,Kato Y,Tanaka M,Yamaguchi H,Kurosawa Y,Yamada H,Fujita N
Sat Nov 26 00:00:00 UTC 2022
Transcriptional and H3K27ac related genome profiles in oral squamous cell carcinoma cells treated with metformin.
Journal of Cancer
Liu S,Shi C,Hou X,Tian X,Li C,Ma X,Wang X,Gao P
Wed Apr 13 00:00:00 UTC 2022
A novel hypoxia-driven gene signature that can predict the prognosis of hepatocellular carcinoma.
Bioengineered
Zeng Z,Lei S,Wang J,Yang Y,Lan J,Tian Q,Chen T,Hao X
Sun May 01 00:00:00 UTC 2022
Identification of HMGB2 associated with proliferation, invasion and prognosis in lung adenocarcinoma via weighted gene co-expression network analysis.
BMC pulmonary medicine
Qiu X,Liu W,Zheng Y,Zeng K,Wang H,Sun H,Dai J
Fri Aug 12 00:00:00 UTC 2022
The Long Non-Coding RNA HOXC-AS3 Promotes Glioma Progression by Sponging miR-216 to Regulate F11R Expression.
Frontiers in oncology
Li Y,Peng L,Cao X,Yang K,Wang Z,Xiao Y,Xiao H,Qian C,Liu H
Wed Apr 13 00:00:00 UTC 2022
NNMT-DNMT1 Axis is Essential for Maintaining Cancer Cell Sensitivity to Oxidative Phosphorylation Inhibition.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wu C,Liu Y,Liu W,Zou T,Lu S,Zhu C,He L,Chen J,Fang L,Zou L,Wang P,Fan L,Wang H,You H,Chen J,Fang JY,Jiang C,Shi Y
Wed Nov 16 00:00:00 UTC 2022
LARP6 Regulates Keloid Fibroblast Proliferation, Invasion, and Ability to Synthesize Collagen.
The Journal of investigative dermatology
Chen L,Su Y,Yin B,Li S,Cheng X,He Y,Jia C
Thu Sep 01 00:00:00 UTC 2022
MiR-1306-5p promotes cell proliferation and inhibits cell apoptosis in acute myeloid leukemia by downregulating PHF6 expression.
Leukemia research
Gao X,Fan S,Zhang X
Thu Sep 01 00:00:00 UTC 2022
Cardiotoxicity of patulin was found in H9c2 cells.
Toxicon : official journal of the International Society on Toxinology
Zhang B,Liang H,Huang K,Li J,Xu D,Huang C,Li Y
Tue Feb 01 00:00:00 UTC 2022
LipoxinA4 analog BML-111 protects podocytes cultured in high-glucose medium against oxidative injury via activating Nrf2 pathway.
International immunopharmacology
Hao H,Xie F,Xu F,Wang Q,Wu Y,Zhang D
Sat Oct 01 00:00:00 UTC 2022
San-Wu-Huang-Qin decoction attenuates tumorigenesis and mucosal barrier impairment in the AOM/DSS model by targeting gut microbiome.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhou Y,Feng Y,Cen R,Hou X,Yu H,Sun J,Zhou L,Ji Q,Zhao L,Wang Y,Li Q
Fri Apr 01 00:00:00 UTC 2022
Morchella esculenta polysaccharide attenuate obesity, inflammation and modulate gut microbiota.
AMB Express
Rehman AU,Khan AI,Xin Y,Liang W
Sat Sep 03 00:00:00 UTC 2022
New Deferric Amine Compounds Efficiently Chelate Excess Iron to Treat Iron Overload Disorders and to Prevent Ferroptosis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Feng W,Xiao Y,Zhao C,Zhang Z,Liu W,Ma J,Ganz T,Zhang J,Liu S
Sat Oct 01 00:00:00 UTC 2022
Yajieshaba prevents lipopolysaccharide-induced intestinal barrier injuryanti-inflammatory and anti-apoptosis.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan
Liping Y,Xinglin YU,Chao Z,Pu C,Xiaohua D
Sat Oct 01 00:00:00 UTC 2022
Yajieshaba prevents lipopolysaccharide-induced intestinal barrier injuryanti-inflammatory and anti-apoptosis.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan
Liping Y,Xinglin YU,Chao Z,Pu C,Xiaohua D
Sat Oct 01 00:00:00 UTC 2022
Grifola frondosa Polysaccharide Ameliorates Early Diabetic Nephropathy by Suppressing the TLR4/NF-κB Pathway.
Applied biochemistry and biotechnology
Jiang T,Shen S,Wang L,Zhao M,Li Y,Huang S
Thu Sep 01 00:00:00 UTC 2022
Corn oligopeptides inhibit Akt/NF-κB signaling pathway and inflammatory factors to ameliorate CCl4 -induced hepatic fibrosis in mice.
Journal of food biochemistry
Feng XW,Cheng QL,Fang L,Liu WY,Liu LW,Sun CQ,Lu ZH,Li GM,Gu RZ
Mon Aug 01 00:00:00 UTC 2022
Whole-exome sequencing and functional validation reveal a rare missense variant in MMP7 that confers ovarian endometriosis risk.
Human molecular genetics
Liu F,Zhou J,Zhang X,Fang S,Liu R,Chen G,Luo Y,Zhang Z,Cheng Y,Wang L,Guo J,Zou Y
Wed Aug 17 00:00:00 UTC 2022
HBO Alleviates Neural Stem Cell Pyroptosis via lncRNA-H19/miR-423-5p/NLRP3 Axis and Improves Neurogenesis after Oxygen Glucose Deprivation.
Oxidative medicine and cellular longevity
Ye Y,Feng Z,Tian S,Yang Y,Jia Y,Wang G,Wang J,Bai W,Li J,He X
Fri Mar 18 00:00:00 UTC 2022
NCAPG Promotes Pulmonary Artery Smooth Muscle Cell Proliferation as a Promising Therapeutic Target of Idiopathic Pulmonary Hypertension: Bioinformatics Analysis and Experiment Verification.
International journal of molecular sciences
Fu B,Li Y,Shi X,Liu P,Zhang Y,Tian H
Tue Oct 04 00:00:00 UTC 2022
Receptor-Interacting Protein Kinase 3 Inhibition Relieves Mechanical Allodynia and Suppresses NLRP3 Inflammasome and NF-κB in a Rat Model of Spinal Cord Injury.
Frontiers in molecular neuroscience
Xue S,Cao ZX,Wang JN,Zhao QX,Han J,Yang WJ,Sun T
Sat May 07 00:00:00 UTC 2022
BRD7 restrains TNF-α-induced proliferation and migration of airway smooth muscle cells by inhibiting notch signaling.
Experimental lung research
Li H,Yang T,Chen T,Liu Y,Pang Y,Yang L
Tue Sep 13 00:00:00 UTC 2022
Palmitoylation of MDH2 by ZDHHC18 activates mitochondrial respiration and accelerates ovarian cancer growth.
Science China. Life sciences
Pei X,Li KY,Shen Y,Li JT,Lei MZ,Fang CY,Lu HJ,Yang HJ,Wen W,Yin M,Qu J,Lei QY
Sat Oct 01 00:00:00 UTC 2022
Protective effect of Astragaloside IV on chronic intermittent hypoxia-induced vascular endothelial dysfunction through the calpain-1/SIRT1/AMPK signaling pathway.
Frontiers in pharmacology
Zhao F,Meng Y,Wang Y,Fan S,Liu Y,Zhang X,Ran C,Wang H,Lu M
Sat Aug 20 00:00:00 UTC 2022
A prognostic signature consisting of metabolism-related genes and SLC17A4 serves as a potential biomarker of immunotherapeutic prediction in prostate cancer.
Frontiers in immunology
Li H,Gu J,Tian Y,Li S,Zhang H,Dai Z,Wang Z,Zhang N,Peng R
Fri Nov 04 00:00:00 UTC 2022
Protective role of selenium on MPP+ and homocysteine-induced TRPM2 channel activation in SH-SY5Y cells.
Journal of receptor and signal transduction research
Yıldızhan K,Nazıroğlu M
Mon Aug 01 00:00:00 UTC 2022
Reduced binding activity of vaccine serum to omicron receptor-binding domain.
Frontiers in immunology
Li M,Weng S,Wang Q,Yang Z,Wang X,Yin Y,Zhou Q,Zhang L,Tao F,Li Y,Jia M,Yang L,Xin X,Li H,Kang L,Wang Y,Wang T,Li S,Kong L
Tue Aug 16 00:00:00 UTC 2022
Bu-Gu-Sheng-Sui decoction promotes osteogenesis via activating the ERK/Smad signaling pathways.
Frontiers in pharmacology
Liu N,Qi B,Zhang Y,Fang S,Sun C,Li Q,Wei X
Tue Sep 13 00:00:00 UTC 2022
Sheng Mai San ameliorated heat stress-induced liver injury via regulating energy metabolism and AMPK/Drp1-dependent autophagy process.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Zhang X,Jia Y,Yuan Z,Wen Y,Zhang Y,Ren J,Ji P,Yao W,Hua Y,Wei Y
Tue Mar 01 00:00:00 UTC 2022
Downregulation of NCL attenuates tumor formation and growth in HeLa cells by targeting the PI3K/AKT pathway.
Cancer medicine
Ying J,Pan R,Tang Z,Zhu J,Ren P,Lou Y,Zhang E,Huang D,Hu P,Li D,Bao Q,Li P
Tue Mar 01 00:00:00 UTC 2022
Naringin regulates mitochondrial dynamics to protect against acetaminophen-induced hepatotoxicity by activating the AMPK/Nrf2 signaling pathway in vitro.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas
Wu Q,Yu P,Bi Y,Li Z,Guo W,Chen Y,Duan Z
Tue Oct 25 00:00:00 UTC 2022
A comprehensive analysis on the relationship between BDE-209 exposure and erectile dysfunction.
Chemosphere
Zhou X,Song L,Cong R,Luan J,Zhou X,Wang Y,Yao L,Zhang X,Ren X,Zhang T,Yu M,Song N
Thu Dec 01 00:00:00 UTC 2022
Tumor-promoting mechanisms of macrophage-derived extracellular vesicles-enclosed microRNA-660 in breast cancer progression.
Breast cancer research and treatment
Li C,Li R,Hu X,Zhou G,Jiang G
Fri Apr 01 00:00:00 UTC 2022
Realgar Alleviated Neuroinflammation Induced by High Protein and High Calorie Diet in Rats via the Microbiota-Gut-Brain Axis.
Nutrients
Feng C,Li A,Yin C,Wang S,Jin W,Liu Y,Huo T,Jiang H
Fri Sep 23 00:00:00 UTC 2022
Quercetin Attenuates Osteoporosis in Orchiectomy Mice by Regulating Glucose and Lipid Metabolism via the GPRC6A/AMPK/mTOR Signaling Pathway.
Frontiers in endocrinology
Sun J,Pan Y,Li X,Wang L,Liu M,Tu P,Wu C,Xiao J,Han Q,Da W,Ma Y,Guo Y
Tue May 17 00:00:00 UTC 2022
A comprehensive analysis of age-related metabolomics and transcriptomics reveals metabolic alterations in rat bone marrow mesenchymal stem cells.
Aging
Yu X,Sun H,Gao X,Zhang C,Sun Y,Wang H,Zhang H,Shi Y,He X
Sun Jan 30 00:00:00 UTC 2022
Liraglutide Attenuates Hepatic Ischemia-Reperfusion Injury by Modulating Macrophage Polarization.
Frontiers in immunology
Li SL,Wang ZM,Xu C,Che FH,Hu XF,Cao R,Xie YN,Qiu Y,Shi HB,Liu B,Dai C,Yang J
Mon Apr 25 00:00:00 UTC 2022
A Dual Anti-Inflammatory and Anti-Proliferative 3-Styrylchromone Derivative Synergistically Enhances the Anti-Cancer Effects of DNA-Damaging Agents on Colon Cancer Cells by Targeting HMGB1-RAGE-ERK1/2 Signaling.
International journal of molecular sciences
Tanuma SI,Oyama T,Okazawa M,Yamazaki H,Takao K,Sugita Y,Amano S,Abe T,Sakagami H
Tue Mar 22 00:00:00 UTC 2022
Lupeol triggers oxidative stress, ferroptosis, apoptosis and restrains inflammation in nasopharyngeal carcinoma via AMPK/NF-κB pathway.
Immunopharmacology and immunotoxicology
Zhou JC,Wu B,Zhang JJ,Zhang W
Mon Aug 01 00:00:00 UTC 2022
Identification of PDCD2 as a Candidate Target of Andrographolide That Arrests the Tumor Cell Cycle by Human Proteome-Scale Screening.
ACS pharmacology & translational science
Wang L,Li C,Chen P,Liu C,Cui Z,Deng S,Yang H
Fri Jul 08 00:00:00 UTC 2022
Succinate communicates pro-inflammatory signals to the host and regulates bile acid enterohepatic metabolism in a pig model.
Food & function
Li X,Ren Y,Huang G,Zhang R,Zhang Y,Zhu W,Yu K
Mon Oct 31 00:00:00 UTC 2022
Geraniin Protects against Cerebral Ischemia/Reperfusion Injury by Suppressing Oxidative Stress and Neuronal Apoptosis via Regulation of the Nrf2/HO-1 Pathway.
Oxidative medicine and cellular longevity
Yang Y,He B,Zhang X,Yang R,Xia X,Chen L,Li R,Shen Z,Chen P
Thu Mar 17 00:00:00 UTC 2022
N-acetyldopamine dimer inhibits neuroinflammation through the TLR4/NF-κB and NLRP3/Caspase-1 pathways.
Acta biochimica et biophysica Sinica
Huang L,Gong L,Huo X,Lei L,Zhang Q,Hu Y,Kuang Q,Gui Y,Dai Y,Gu Y,Deng Y,Wang D,Guo D
Thu Aug 25 00:00:00 UTC 2022
JMJD1C Regulates Megakaryopoiesis in In Vitro Models through the Actin Network.
Cells
Wang J,Liu X,Wang H,Qin L,Feng A,Qi D,Wang H,Zhao Y,Kong L,Wang H,Wang L,Hu Z,Xu X
Fri Nov 18 00:00:00 UTC 2022
Selenoprotein K contributes to CD36 subcellular trafficking in hepatocytes by accelerating nascent COPII vesicle formation and aggravates hepatic steatosis.
Redox biology
You M,Wu F,Gao M,Chen M,Zeng S,Zhang Y,Zhao W,Li D,Wei L,Ruan XZ,Chen Y
Tue Nov 01 00:00:00 UTC 2022
Sertad1 Induces Neurological Injury after Ischemic Stroke via the CDK4/p-Rb Pathway.
Molecules and cells
Li J,Li B,Bu Y,Zhang H,Guo J,Hu J,Zhang Y
Sat Apr 30 00:00:00 UTC 2022
Sertad1 Induces Neurological Injury after Ischemic Stroke via the CDK4/p-Rb Pathway.
Molecules and cells
Li J,Li B,Bu Y,Zhang H,Guo J,Hu J,Zhang Y
Sat Apr 30 00:00:00 UTC 2022
G3BP2, a stress granule assembly factor, is dispensable for spermatogenesis in mice.
PeerJ
Yun D,Zhou L,Shi J,Li X,Wu X,Sun F
Sat Jan 21 00:00:00 UTC 2023
Antidepressant effects of total alkaloids of Fibraurea recisa on improving corticosterone-induced apoptosis of HT-22 cells and chronic unpredictable mild stress-induced depressive-like behaviour in mice.
Pharmaceutical biology
He Z,Yu H,Wu H,Su L,Shi K,Zhao Y,Zong Y,Chen W,Du R
Thu Dec 01 00:00:00 UTC 2022
TRIM36 inhibits tumorigenesis through the Wnt/β-catenin pathway and promotes caspase-dependent apoptosis in hepatocellular carcinoma.
Cancer cell international
Tong Q,Yi M,Kong P,Xu L,Huang W,Niu Y,Gan X,Zhan H,Tian R,Yan D
Tue Sep 06 00:00:00 UTC 2022
African Swine Fever Virus K205R Induces ER Stress and Consequently Activates Autophagy and the NF-κB Signaling Pathway.
Viruses
Wang Q,Zhou L,Wang J,Su D,Li D,Du Y,Yang G,Zhang G,Chu B
Tue Feb 15 00:00:00 UTC 2022
Multifunctional Drug-Loaded Phase-Change Nanoparticles Inhibit the Epithelial-Mesenchymal Transition of Hepatocellular Carcinoma by Affecting the Activity of Activated Hepatic Stellate Cells.
BioMed research international
Zou X,Li T,Mao Y,Zhu M,Chen X,Niu J,Dong T,Jiang J,Yang X
Wed Nov 23 00:00:00 UTC 2022
SNORA70E promotes the occurrence and development of ovarian cancer through pseudouridylation modification of RAP1B and alternative splicing of PARPBP.
Journal of cellular and molecular medicine
Chen S,Li QH,Chen X,Bao HJ,Wu W,Shen F,Lu BF,Jiang RQ,Zong ZH,Zhao Y
Sat Oct 01 00:00:00 UTC 2022
A New Long Noncoding RNA, MAHAT, Inhibits Replication of Porcine Reproductive and Respiratory Syndrome Virus by Recruiting DDX6 To Bind to ZNF34 and Promote an Innate Immune Response.
Journal of virology
Liu Y,Liu X,Bai J,Sun Y,Nauwynck H,Wang X,Yang Y,Jiang P
Wed Sep 28 00:00:00 UTC 2022
Evodiamine as the Active Compound of Evodiae fructus to Inhibit Proliferation and Migration of Prostate Cancer through PI3K/AKT/NF-κB Signaling Pathway.
Disease markers
Lei Y,Chan M,Liu H,Lyu W,Chen L,Zhong Y,Gan H,Wang M,Qi M,Guo Y,Liu J,Zhang E
Fri Jul 29 00:00:00 UTC 2022
Magnoflorine attenuates inflammatory responses in RA by regulating the PI3K/Akt/NF-κB and Keap1-Nrf2/HO-1 signalling pathways in vivo and in vitro.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Shen Y,Fan X,Qu Y,Tang M,Huang Y,Peng Y,Fu Q
Thu Sep 01 00:00:00 UTC 2022
Identification of circular RNA BTBD7_hsa_circ_0000563 as a novel biomarker for coronary artery disease and the functional discovery of BTBD7_hsa_circ_0000563 based on peripheral blood mononuclear cells: a case control study.
Clinical proteomics
Zhou H,Gan X,He S,Wang Y,Zhang S,Chen J,Zhou Y,Hou C,Hua L,Zhang Q,Jia E
Thu Nov 03 00:00:00 UTC 2022
Sevelamer arsenite nanoparticle as a Pi-responsive drug carrier and embolic agent for chemoembolization.
Drug delivery
Bi QC,Tang JJ,Zhao J,Lv YF,Deng ZQ,Chen H,Xu YH,Xie CS,Liang QR,Luo RG,Tang Q
Thu Dec 01 00:00:00 UTC 2022
Pterostilbene and Its Derivative 3'-Hydroxypterostilbene Ameliorated Nonalcoholic Fatty Liver Disease Through Synergistic Modulation of the Gut Microbiota and SIRT1/AMPK Signaling Pathway.
Journal of agricultural and food chemistry
Tsai HY,Shih YY,Yeh YT,Huang CH,Liao CA,Hu CY,Nagabhushanam K,Ho CT,Chen YK
Wed Apr 27 00:00:00 UTC 2022
Trans-anethole Ameliorates Intestinal Injury Through Activation of Nrf2 Signaling Pathway in Subclinical Necrotic Enteritis-Induced Broilers.
Frontiers in veterinary science
Yu C,Tong Y,Li Q,Wang T,Yang Z
Sat May 07 00:00:00 UTC 2022
Mulberry Leaf Flavonoids Inhibit Liver Inflammation in Type 2 Diabetes Rats by Regulating TLR4/MyD88/NF-κB Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Duan Y,Dai H,An Y,Cheng L,Shi L,Lv Y,Li H,Wang C,He C,Zhang H,Huang Y,Fu W,Meng Y,Zhao B
Tue Jul 19 00:00:00 UTC 2022
A marine-derived small molecule induces immunogenic cell death against triple-negative breast cancer through ER stress-CHOP pathway.
International journal of biological sciences
Wen H,Zhong Y,Yin Y,Qin K,Yang L,Li D,Yu W,Yang C,Deng Z,Hong K
Thu May 12 00:00:00 UTC 2022
A multi-target directed ligands strategy for the treatment of Alzheimer's disease: Dimethyl fumarate plus Tranilast modified Dithiocarbate as AChE inhibitor and Nrf2 activator.
European journal of medicinal chemistry
Guo J,Cheng M,Liu P,Cao D,Luo J,Wan Y,Fang Y,Jin Y,Xie SS,Liu J
Tue Nov 15 00:00:00 UTC 2022
Effects of intraperitoneal injection of lipopolysaccharide-induced peripheral inflammation on dopamine neuron damage in rat midbrain.
CNS neuroscience & therapeutics
Yang QY,Li XW,Yang R,Qin TY,Long H,Zhang SB,Zhang F
Sat Oct 01 00:00:00 UTC 2022
Toxoplasma gondii dense granule protein 3 promotes endoplasmic reticulum stress-induced apoptosis by activating the PERK pathway.
Parasites & vectors
Obed C,Wu M,Chen Y,An R,Cai H,Luo Q,Yu L,Wang J,Liu F,Shen J,Du J
Tue Aug 02 00:00:00 UTC 2022
Maltol Promotes Mitophagy and Inhibits Oxidative Stress via the Nrf2/PINK1/Parkin Pathway after Spinal Cord Injury.
Oxidative medicine and cellular longevity
Mao Y,Du J,Chen X,Al Mamun A,Cao L,Yang Y,Mubwandarikwa J,Zaeem M,Zhang W,Chen Y,Dai Y,Xiao J,Ye K
Fri Mar 18 00:00:00 UTC 2022
Chelerythrine inhibits the progression of glioblastoma by suppressing the TGFB1-ERK1/2/Smad2/3-Snail/ZEB1 signaling pathway.
Life sciences
Zhu M,Niu J,Jiang J,Dong T,Chen Y,Yang X,Liu P
Tue Mar 15 00:00:00 UTC 2022
Maternal Inflammation Exaggerates Offspring Susceptibility to Cerebral Ischemia-Reperfusion Injury via the COX-2/PGD2/DP2 Pathway Activation.
Oxidative medicine and cellular longevity
Li Y,Luo W,Zhang J,Luo Y,Han W,Wang H,Xia H,Chen Z,Yang Y,Chen Q,Li H,Yang L,Hu C,Huang H,Peng Z,Tan X,Li M,Yang J
Wed Apr 20 00:00:00 UTC 2022
Preliminary Evaluation of Artificial Intelligence-Based Anti-Hepatocellular Carcinoma Molecular Target Study in Hepatocellular Carcinoma Diagnosis Research.
BioMed research international
Wang Y,Wei C,Deng X,Gao S,Chen J
Wed Oct 05 00:00:00 UTC 2022
ASFV pD345L protein negatively regulates NF-κB signalling by inhibiting IKK kinase activity.
Veterinary research
Chen H,Wang Z,Gao X,Lv J,Hu Y,Jung YS,Zhu S,Wu X,Qian Y,Dai J
Sat Apr 23 00:00:00 UTC 2022
NAD(P)H: quinone oxidoreductase 1 attenuates oxidative stress and apoptosis by regulating Sirt1 in diabetic nephropathy.
Journal of translational medicine
Qiu D,Song S,Wang Y,Bian Y,Wu M,Wu H,Shi Y,Duan H
Fri Jan 28 00:00:00 UTC 2022
TRIM47 promotes ovarian cancer cell proliferation, migration, and invasion by activating STAT3 signaling.
Clinics (Sao Paulo, Brazil)
Wang X,Fu Y,Xing Y
Mon Oct 31 00:00:00 UTC 2022
Liver fibrosis alters the molecular structures of hepatic glycogen.
Carbohydrate polymers
Wan Y,Hu Z,Liu Q,Wang L,Sullivan MA,Gilbert RG
Tue Feb 15 00:00:00 UTC 2022
Organophosphate flame retardant TDCPP: A risk factor for renal cancer?
Chemosphere
Zhou X,Zhou X,Yao L,Zhang X,Cong R,Luan J,Zhang T,Song N
Sat Oct 01 00:00:00 UTC 2022
Acetyl-L-carnitine improves erectile function in bilateral cavernous nerve injury rats via promoting cavernous nerve regeneration.
Andrology
Wang J,Song J,Song G,Feng Y,Pan J,Yang X,Xin Z,Hu P,Sun T,Liu K,Xu W,Wang T,Wang S,Liu J,Ruan Y
Fri Jul 01 00:00:00 UTC 2022
A single-beam of light priming the immune responses and boosting cancer photoimmunotherapy.
Journal of controlled release : official journal of the Controlled Release Society
Sun Y,Han R,Wang J,Qin Y,Ren Z,Feng X,Liu Q,Wang X
Sat Oct 01 00:00:00 UTC 2022
Deciphering the tumor microenvironment cell-infiltrating landscape reveals microenvironment subtypes and therapeutic potentials for nonsquamous NSCLC.
JCI insight
Chen H,Zhang T,Zhang Y,Wu H,Fang Z,Liu Y,Chen Y,Wang Z,Jia S,Ji X,Shang L,Du F,Liu J,Lu M,Chong W
Wed Jun 22 00:00:00 UTC 2022
Melatonin promotes sirtuin 1 expression and inhibits IRE1α-XBP1S-CHOP to reduce endoplasmic reticulum stress-mediated apoptosis in chondrocytes.
Frontiers in pharmacology
Qin K,Tang H,Ren Y,Yang D,Li Y,Huang W,Wu Y,Yin Z
Tue Aug 30 00:00:00 UTC 2022
DDX47 promotes cell proliferation and migration in lung adenocarcinoma.
Pathology, research and practice
Chen Y,Zeng L,Wang W,Yuan J,Sun R,Yang S
Wed Jun 01 00:00:00 UTC 2022
Immunity of two novel hepatitis C virus polyepitope vaccines.
Vaccine
Feng T,Li M,Zhang L,Li S,Yang Z,Kang L,Guo Y,Kong L,Wang T
Wed Oct 12 00:00:00 UTC 2022
Severe inflammation in new-borns induces long-term cognitive impairment by activation of IL-1β/KCC2 signaling during early development.
BMC medicine
Zhang D,Yang Y,Yang Y,Liu J,Zhu T,Huang H,Zhou C
Wed Jul 27 00:00:00 UTC 2022
U2 small nuclear RNA auxiliary factor 2, transcriptionally activated by the transcription factor Dp-1/E2F transcription factor 1 complex, enhances the growth and aerobic glycolysis of leiomyosarcoma cells.
Bioengineered
Li Y,Chen S,Zhang X,Zhuo N
Fri Apr 01 00:00:00 UTC 2022
ChemR23 signaling ameliorates cognitive impairments in diabetic mice via dampening oxidative stress and NLRP3 inflammasome activation.
Redox biology
Zhang J,Liu L,Zhang Y,Yuan Y,Miao Z,Lu K,Zhang X,Ni R,Zhang H,Zhao Y,Wang X
Thu Dec 01 00:00:00 UTC 2022
Ghrelin ameliorates diabetes-associated behavioral deficits and NLRP3 inflammasome activation via autophagic flux enhancement.
Pharmacological research
Han W,Cui C,Zhang H,Guo Y,Xie D,Zhang W,Wang C,Yang M,Jiang P
Sun May 01 00:00:00 UTC 2022
PCV2 and PRV Coinfection Induces Endoplasmic Reticulum Stress via PERK-eIF2α-ATF4-CHOP and IRE1-XBP1-EDEM Pathways.
International journal of molecular sciences
Chen S,Li X,Zhang X,Niu G,Yang L,Ji W,Zhang L,Ren L
Tue Apr 19 00:00:00 UTC 2022
Salecan ameliorates liver injury by regulating gut microbiota and its metabolites.
Food & function
Li P,Liu Y,Zhao J,Pan W,He Y,Fu S,Liu Y,Xu YJ
Mon Nov 14 00:00:00 UTC 2022
Decitabine attenuates ischemic stroke by reducing astrocytes proliferation in rats.
PloS one
Zhang Q,Li D,Zhao H,Zhang X
Thu Aug 04 00:00:00 UTC 2022
Anti-photoaging effects of flexible nanoliposomes encapsulated Moringa oleifera Lam. isothiocyanate in UVB-induced cell damage in HaCaT cells.
Drug delivery
Wang Y,Ouyang Q,Chang X,Yang M,He J,Tian Y,Sheng J
Thu Dec 01 00:00:00 UTC 2022
Study on the mechanism of scutellarin's protective effect against ZEA-induced mouse ovarian granulosa cells injury.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Yi Y,Hao Z,Sun P,Fan K,Yin W,Guo J,Zheng X,Sun N,Li H
Thu Dec 01 00:00:00 UTC 2022
Ibandronate promotes autophagy by inhibiting Rac1-mTOR signaling pathway in vitro and in vivo.
Cell death discovery
Han J,Yang J,Wang Q,Yin X,Sun Z,Huang C,Chen G,Zheng L,Jiang D
Sat Apr 09 00:00:00 UTC 2022
ABHD16A Negatively Regulates the Palmitoylation and Antiviral Function of IFITM Proteins.
mBio
Shi X,Li X,Xu Z,Shen L,Ding Y,Chen S,Mao L,Liu W,Xu J
Tue Dec 20 00:00:00 UTC 2022
ABHD16A Negatively Regulates the Palmitoylation and Antiviral Function of IFITM Proteins.
mBio
Shi X,Li X,Xu Z,Shen L,Ding Y,Chen S,Mao L,Liu W,Xu J
Tue Dec 20 00:00:00 UTC 2022
ABHD16A Negatively Regulates the Palmitoylation and Antiviral Function of IFITM Proteins.
mBio
Shi X,Li X,Xu Z,Shen L,Ding Y,Chen S,Mao L,Liu W,Xu J
Tue Dec 20 00:00:00 UTC 2022
ABHD16A Negatively Regulates the Palmitoylation and Antiviral Function of IFITM Proteins.
mBio
Shi X,Li X,Xu Z,Shen L,Ding Y,Chen S,Mao L,Liu W,Xu J
Tue Dec 20 00:00:00 UTC 2022
Cystathionine β-Synthase Regulates the Proliferation, Migration, and Invasion of Thyroid Carcinoma Cells.
Oxidative medicine and cellular longevity
Jiang QY,Li JM,Jing MR,Zhang YX,Zhang QQ,Cai CB,Wang D,Qi HW,Li T,Li YZ,Ji XY,Wu DD
Fri Jul 08 00:00:00 UTC 2022
Alpha-Asaronol Alleviates Dysmyelination by Enhancing Glutamate Transport Through the Activation of PPARγ-GLT-1 Signaling in Hypoxia-Ischemia Neonatal Rats.
Frontiers in pharmacology
Ge Y,Zhen F,Liu Z,Feng Z,Wang G,Zhang C,Wang X,Sun Y,Zheng X,Bai Y,Yao R
Wed Apr 13 00:00:00 UTC 2022
Substitution of SERCA2 Cys674 accelerates aortic aneurysm by inducing endoplasmic reticulum stress and promoting cell apoptosis.
British journal of pharmacology
Wang L,Yang Z,Wang S,Que Y,Shu X,Wu F,Liu G,Li S,Hu P,Chen H,Shi J,Tong X
Thu Sep 01 00:00:00 UTC 2022
Sestrin2 remedies podocyte injury via orchestrating TSP-1/TGF-β1/Smad3 axis in diabetic kidney disease.
Cell death & disease
Song S,Shi C,Bian Y,Yang Z,Mu L,Wu H,Duan H,Shi Y
Sat Jul 30 00:00:00 UTC 2022
RNA-m6A modification of HDGF mediated by Mettl3 aggravates the progression of atherosclerosis by regulating macrophages polarization via energy metabolism reprogramming.
Biochemical and biophysical research communications
Zheng L,Chen X,Yin Q,Gu J,Chen J,Chen M,Zhang Y,Dong M,Jiang H,Yin N,Chen H,Li X
Tue Dec 20 00:00:00 UTC 2022
Grape Seed Proanthocyanidins Exert a Neuroprotective Effect by Regulating Microglial M1/M2 Polarisation in Rats with Spinal Cord Injury.
Mediators of inflammation
Liu WZ,Ma ZJ,Kang JH,Lin AX,Wang ZH,Chen HW,Guo XD,He XG,Kang XW
Tue Aug 16 00:00:00 UTC 2022
Oroxin A ameliorates the oleic acid-induced A549 cell injury through the suppression of pyroptosis and degradation of alveolar surfactant.
Anais da Academia Brasileira de Ciencias
Huang J,Chen C,Xie J,Zhai K,Wei S,Cheng X,Zhang R
Thu Dec 15 00:00:00 UTC 2022
Moxidectin induces autophagy arrest in colorectal cancer.
Medical oncology (Northwood, London, England)
Mao Y,Xie H,Shu D,Cheng L,Lan J,Pan K
Thu Sep 29 00:00:00 UTC 2022
Inhibition of HIF-1α Attenuates Silica-Induced Pulmonary Fibrosis.
International journal of environmental research and public health
Xu X,Li Y,Niu Z,Xia J,Dai K,Wang C,Yao W,Guo Y,Deng X,He J,Deng M,Si H,Hao C
Wed Jun 01 00:00:00 UTC 2022
Transketolase promotes colorectal cancer metastasis through regulating AKT phosphorylation.
Cell death & disease
Li M,Zhao X,Yong H,Xu J,Qu P,Qiao S,Hou P,Li Z,Chu S,Zheng J,Bai J
Wed Feb 02 00:00:00 UTC 2022
Bisphenol A impairs macrophages through inhibiting autophagy via AMPK/mTOR signaling pathway and inducing apoptosis.
Ecotoxicology and environmental safety
Wu M,Cong Y,Wang K,Yu H,Zhang X,Ma M,Duan Z,Pei X
Fri Apr 01 00:00:00 UTC 2022
Corilagin induces apoptosis and inhibits autophagy of HL‑60 cells by regulating miR‑451/HMGB1 axis.
Molecular medicine reports
Jia S,He D,Liang X,Cheng P,Liu J,Chen M,Wang C,Zhang H,Meng C
Sat Jan 01 00:00:00 UTC 2022
Taurine protects R28 cells from hypoxia/re-oxygenation-induced damage via regulation of mitochondrial energy metabolism.
Amino acids
Lu W,Yang Y,Gao S,Wu J,Sun X
Thu Dec 01 00:00:00 UTC 2022
Nicotinamide riboside promotes Mfn2-mediated mitochondrial fusion in diabetic hearts through the SIRT1-PGC1α-PPARα pathway.
Free radical biology & medicine
Hu L,Guo Y,Song L,Wen H,Sun N,Wang Y,Qi B,Liang Q,Geng J,Liu X,Fu F,Li Y
Fri Apr 01 00:00:00 UTC 2022
M1 Macrophages Enhance Survival and Invasion of Oral Squamous Cell Carcinoma by Inducing GDF15-Mediated ErbB2 Phosphorylation.
ACS omega
Lv C,Li S,Zhao J,Yang P,Yang C
Tue Apr 05 00:00:00 UTC 2022
Low expression of microRNA-125b enhances the expression of STAT3 and contributes to cholesteatoma growth.
Archives of medical science : AMS
Zang J,Yang B,Feng S,Jiang X
Sat Dec 03 00:00:00 UTC 2022
The Effect of Circumscribed Exposure to the Pan-Aurora Kinase Inhibitor VX-680 on Proliferating Euploid Cells.
International journal of molecular sciences
Liu X,Shi Q,Choudhry N,Zhang T,Liu H,Zhang S,Zhang J,Yang D
Tue Oct 11 00:00:00 UTC 2022
HNRNPK/CLCN3 axis facilitates the progression of LUAD through CAF-tumor interaction.
International journal of biological sciences
Li Y,Yang Y,Ma Q,Cheng H,Wang H,Ma C,Li F,Zhao S,Li X,Qi Y,Gu Z
Tue Nov 29 00:00:00 UTC 2022
HNRNPK/CLCN3 axis facilitates the progression of LUAD through CAF-tumor interaction.
International journal of biological sciences
Li Y,Yang Y,Ma Q,Cheng H,Wang H,Ma C,Li F,Zhao S,Li X,Qi Y,Gu Z
Tue Nov 29 00:00:00 UTC 2022
Inhibition of APE1 Expression Enhances the Antitumor Activity of Olaparib in Triple-Negative Breast Cancer.
Evidence-based complementary and alternative medicine : eCAM
Jian D,Li XM,Dai N,Liang DD,Zhang G,Mao CY,Wang D,Song GB,Li MX,Luo H
Sat Jul 16 00:00:00 UTC 2022
Glycogen metabolism reprogramming promotes inflammation in coal dust-exposed lung.
Ecotoxicology and environmental safety
Wang W,Mu M,Zou Y,Deng S,Lu Y,Li Q,Li Z,Tao H,Wang Y,Tao X
Thu Sep 01 00:00:00 UTC 2022
Poldip2 knockdown protects against lipopolysaccharide-induced acute lung injury via Nox4/Nrf2/NF-κB signaling pathway.
Frontiers in pharmacology
Wang Y,Wang W,Zhou S,Wang Y,Cudjoe O,Cha Y,Wang C,Cao X,Liu W,Jin K
Tue Sep 20 00:00:00 UTC 2022
Ubiquitin-specific protease 47 regulates intestinal inflammation through deubiquitination of TRAF6 in epithelial cells.
Science China. Life sciences
Lei H,Yang L,Xu H,Wang Z,Li X,Liu M,Wu Y
Mon Aug 01 00:00:00 UTC 2022
Nuciferine Inhibits TMEM16A in Dietary Adjuvant Therapy for Lung Cancer.
Journal of agricultural and food chemistry
Bai X,Liu X,Li S,An H,Kang X,Guo S
Wed Mar 30 00:00:00 UTC 2022
An oleanolic acid derivative, K73-03, inhibits pancreatic cancer cells proliferation in vitro and in vivo via blocking EGFR/Akt pathway.
Cell biology international
Zhou Z,Dong Y,Li N,Niu M,Wang S,Zhou Y,Sun Z,Chu P,Tang Z
Tue Nov 01 00:00:00 UTC 2022
Calycosin suppresses colorectal cancer progression by targeting ERβ, upregulating PTEN, and inhibiting PI3K/Akt signal pathway.
Cell biology international
Zhu L,Liu S,Liao YF,Sheng YM,He JC,Cai ZX,Man Q,Wu YY
Thu Sep 01 00:00:00 UTC 2022
Tension of plus-end tracking protein Clip170 confers directionality and aggressiveness during breast cancer migration.
Cell death & disease
Hu Y,Xie Q,Wu X,Liu W,Li D,Li C,Zhao W,Chen L,Zheng Z,Li G,Guo J
Sat Oct 08 00:00:00 UTC 2022
Activation of the NLRP3 inflammasome by RAC1 mediates a new mechanism in diabetic nephropathy.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Ying C,Zhou Z,Dai J,Wang M,Xiang J,Sun D,Zhou X
Tue Feb 01 00:00:00 UTC 2022
Dye-Sensitized Rare Earth Nanoparticles with Up/Down Conversion Luminescence for On-Demand Gas Therapy of Glioblastoma Guided by NIR-II Fluorescence Imaging.
Advanced healthcare materials
Liu Z,Yun B,Han Y,Jiang Z,Zhu H,Ren F,Li Z
Tue Feb 01 00:00:00 UTC 2022
Essential role of MALAT1 in reducing traumatic brain injury.
Neural regeneration research
Wu N,Cheng CJ,Zhong JJ,He JC,Zhang ZS,Wang ZG,Sun XC,Liu H
Mon Aug 01 00:00:00 UTC 2022
Glucagon-like Peptide 1 Receptor Activation Inhibits Microglial Pyroptosis via Promoting Mitophagy to Alleviate Depression-like Behaviors in Diabetic Mice.
Nutrients
Yang F,Wang X,Qi J,Zhang K,Jiang Y,Feng B,Lv T,Yang L,Yang Q,Zhao M,Liu S,Ma X
Wed Dec 21 00:00:00 UTC 2022
IL-22 Protects against Biliary Ischemia-Reperfusion Injury after Liver Transplantation via Activating STAT3 and Reducing Apoptosis and Oxidative Stress Levels In Vitro and In Vivo.
Oxidative medicine and cellular longevity
Bai Y,Wu H,Zhang J,Zhang S,Zhang Z,Wang H,Zhang Y,Shen Z
Mon May 23 00:00:00 UTC 2022
Paeoniflorin-6'-o-benzene sulfonate ameliorates the progression of adjuvant-induced arthritis by inhibiting the interaction between Ahr and GRK2 of fibroblast-like synoviocytes.
International immunopharmacology
Zhang BJ,Wang YY,Jia CY,Li SS,Wang XW,Xu Y,Chen AY,Xu HP,Wang C,Yang ZY,Wei W,Chang Y
Fri Jul 01 00:00:00 UTC 2022
Weighted gene co-expression network indicates that the DYNLL2 is an important regulator of chicken breast muscle development and is regulated by miR-148a-3p.
BMC genomics
Li Y,Yuan P,Fan S,Zhai B,Jin W,Li D,Li H,Sun G,Han R,Liu X,Tian Y,Li G,Kang X
Mon Apr 04 00:00:00 UTC 2022
Genistein Alleviates Oxidative Stress and Inflammation in the Hypothalamic Paraventricular Nucleus by Activating the Sirt1/Nrf2 Pathway in High Salt-Induced Hypertension.
Cardiovascular toxicology
Niu LG,Sun N,Liu KL,Su Q,Qi J,Fu LY,Xin GR,Kang YM
Tue Nov 01 00:00:00 UTC 2022
Identification of Trovafloxacin, Ozanimod, and Ozenoxacin as Potent c-Myc G-Quadruplex Stabilizers to Suppress c-Myc Transcription and Myeloma Growth.
Molecular informatics
Zhang J,Wang T,Geng X,Liu L,Gao J
Sat Oct 01 00:00:00 UTC 2022
Endothelial cells regulated by RNF20 orchestrate the proliferation and differentiation of neural precursor cells during embryonic development.
Cell reports
Zhang M,Su L,Wang W,Li C,Liang Q,Ji F,Jiao J
Tue Sep 13 00:00:00 UTC 2022
A novel protein encoded by ZCRB1-induced circHEATR5B suppresses aerobic glycolysis of GBM through phosphorylation of JMJD5.
Journal of experimental & clinical cancer research : CR
Song J,Zheng J,Liu X,Dong W,Yang C,Wang D,Ruan X,Zhao Y,Liu L,Wang P,Zhang M,Liu Y
Tue May 10 00:00:00 UTC 2022
Genetic Deficiency of p53 Leads to Structural, Functional, and Synaptic Deficits in Primary Somatosensory Cortical Neurons of Adult Mice.
Frontiers in molecular neuroscience
Kuang H,Liu T,Jiao C,Wang J,Wu S,Wu J,Peng S,Davidson AM,Zeng SX,Lu H,Mostany R
Fri Apr 29 00:00:00 UTC 2022
CTNND1 variants cause familial exudative vitreoretinopathy through the Wnt/cadherin axis.
JCI insight
Yang M,Li S,Huang L,Zhao R,Dai E,Jiang X,He Y,Lu J,Peng L,Liu W,Zhang Z,Jiang D,Zhang Y,Jiang Z,Yang Y,Zhao P,Zhu X,Ding X,Yang Z
Fri Jul 22 00:00:00 UTC 2022
Myeloid peroxisome proliferator-activated receptor α deficiency accelerates liver regeneration via IL-6/STAT3 pathway after 2/3 partial hepatectomy in mice.
Hepatobiliary surgery and nutrition
Xie G,Song Y,Li N,Zhang Z,Wang X,Liu Y,Jiao S,Wei M,Yu B,Wang Y,Wang H,Qu A
Fri Apr 01 00:00:00 UTC 2022
Mettl14-Mediated m6A Modification Is Essential for Germinal Center B Cell Response.
Journal of immunology (Baltimore, Md. : 1950)
Huang H,Zhang G,Ruan GX,Li Y,Chen W,Zou J,Zhang R,Wang J,Ji SJ,Xu S,Ou X
Fri Apr 15 00:00:00 UTC 2022
TFRC upregulation promotes ferroptosis in CVB3 infection via nucleus recruitment of Sp1.
Cell death & disease
Yi L,Hu Y,Wu Z,Li Y,Kong M,Kang Z,Zuoyuan B,Yang Z
Mon Jul 11 00:00:00 UTC 2022
Long Noncoding RNA Metastasis-Associated Lung Adenocarcinoma Transcript 1 in Extracellular Vesicles Promotes Hepatic Stellate Cell Activation, Liver Fibrosis and β-Catenin Signaling Pathway.
Frontiers in physiology
Wang T,Zhang C,Meng X,Zhu B,Wang S,Yuan W,Zhang S,Xu J,Zhang C
Fri Mar 04 00:00:00 UTC 2022
Protection of Human Lens Epithelial Cells from Oxidative Stress Damage and Cell Apoptosis by KGF-2 through the Akt/Nrf2/HO-1 Pathway.
Oxidative medicine and cellular longevity
Liu S,Jin Z,Xia R,Zheng Z,Zha Y,Wang Q,Wan X,Yang H,Cai J
Thu Mar 17 00:00:00 UTC 2022
The hsa-miR-214-3p/ATGL axis regulates aberrant lipolysis to promote acute myeloid leukemia progression via PPARα in vitro.
Biochemical and biophysical research communications
Li Y,Yuan S,Wu W,Zhou J,Zhang P,Li D,Zhang Y,Lou S
Sat Jun 11 00:00:00 UTC 2022
Alzheimer's risk factor FERMT2 promotes the progression of colorectal carcinoma via Wnt/β-catenin signaling pathway and contributes to the negative correlation between Alzheimer and cancer.
PloS one
Xia W,Gao Z,Jiang X,Jiang L,Qin Y,Zhang D,Tian P,Wang W,Zhang Q,Zhang R,Zhang N,Xu S
Wed Dec 21 00:00:00 UTC 2022
Hydroxysafflor Yellow A Blocks HIF-1α Induction of NOX2 and Protects ZO-1 Protein in Cerebral Microvascular Endothelium.
Antioxidants (Basel, Switzerland)
Li Y,Liu XT,Zhang PL,Li YC,Sun MR,Wang YT,Wang SP,Yang H,Liu BL,Wang M,Gao W,Li P
Thu Apr 07 00:00:00 UTC 2022
The BDNF-TrkB signaling pathway in the rostral anterior cingulate cortex is involved in the development of pain aversion in rats with bone cancer via NR2B and ERK-CREB signaling.
Brain research bulletin
Li J,Wang X,Wang H,Wang R,Guo Y,Xu L,Zhang G,Wu J,Wang G
Fri Jul 01 00:00:00 UTC 2022
Cinobufagin induces acute promyelocytic leukaemia cell apoptosis and PML-RARA degradation in a caspase-dependent manner by inhibiting the β-catenin signalling pathway.
Pharmaceutical biology
Bian Y,Xue M,Guo X,Jiang W,Zhao Y,Zhang Z,Wang X,Hu Y,Zhang Q,Dun W,Zhang L
Thu Dec 01 00:00:00 UTC 2022
Novel LncRNA ZFHX4-AS1 as a Potential Prognostic Biomarker That Affects the Immune Microenvironment in Ovarian Cancer.
Frontiers in oncology
Wang X,Wang Y,Sun F,Xu Y,Zhang Z,Yang C,Zhang L,Lou G
Sun Jul 31 00:00:00 UTC 2022
Bile Duct Ligation Impairs Function and Expression of Mrp1 at Rat Blood-Retinal Barrier via Bilirubin-Induced P38 MAPK Pathway Activations.
International journal of molecular sciences
Li P,Yang Y,Lin Z,Hong S,Jiang L,Zhou H,Yang L,Zhu L,Liu X,Liu L
Mon Jul 11 00:00:00 UTC 2022
Overexpression of heat shock protein 70 induces apoptosis of intestinal epithelial cells in heat-stressed pigs: A proteomics approach.
Journal of thermal biology
Yong Y,Li J,Yu T,Fang B,Liu X,Yu Z,Ma X,Gooneratne R,Abd El-Aty AM,Ju X
Mon Aug 01 00:00:00 UTC 2022
The hnRNPK/A1/R/U Complex Regulates Gene Transcription and Translation and is a Favorable Prognostic Biomarker for Human Colorectal Adenocarcinoma.
Frontiers in oncology
Li Y,Wang H,Wan J,Ma Q,Qi Y,Gu Z
Tue Jul 26 00:00:00 UTC 2022
Endothelial Cells Mediated by UCP2 Control the Neurogenic-to-Astrogenic Neural Stem Cells Fate Switch During Brain Development.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wang W,Su L,Wang Y,Li C,Ji F,Jiao J
Wed Jun 01 00:00:00 UTC 2022
HPV16 E6/E7 stabilize PGK1 protein by reducing its poly-ubiquitination in cervical cancer.
Cell biology international
Liu S,Song L,Yao H,Zhang L
Tue Mar 01 00:00:00 UTC 2022
HPV16 E6/E7 stabilize PGK1 protein by reducing its poly-ubiquitination in cervical cancer.
Cell biology international
Liu S,Song L,Yao H,Zhang L
Tue Mar 01 00:00:00 UTC 2022
High expression of Sam68 contributes to metastasis by regulating vimentin expression and a motile phenotype in oral squamous cell carcinoma.
Oncology reports
Komiyama T,Kuroshima T,Sugasawa T,Fujita SI,Ikami Y,Hirai H,Tsushima F,Michi Y,Kayamori K,Higashino F,Harada H
Sat Oct 01 00:00:00 UTC 2022
Effect of transferrin glycation induced by high glucose on HK-2 cells in vitro.
Frontiers in endocrinology
Ma Y,Zhou Q,Zhao P,Lv X,Gong C,Gao J,Liu J
Tue Feb 14 00:00:00 UTC 2023
Macrophage SCAP Contributes to Metaflammation and Lean NAFLD by Activating STING-NF-κB Signaling Pathway.
Cellular and molecular gastroenterology and hepatology
Huang X,Yao Y,Hou X,Wei L,Rao Y,Su Y,Zheng G,Ruan XZ,Li D,Chen Y
Mon Jun 20 00:00:00 UTC 2022
Upregulated LINC01088 facilitates malignant phenotypes and immune escape of colorectal cancer by regulating microRNAs/G3BP1/PD-L1 axis.
Journal of cancer research and clinical oncology
Li C,Pan B,Wang X,Liu X,Qin J,Gao T,Sun H,Pan Y,Wang S
Mon Aug 01 00:00:00 UTC 2022
Assembly of Celastrol to Zeolitic Imidazolate Framework-8 by Coordination as a Novel Drug Delivery Strategy for Cancer Therapy.
Pharmaceuticals (Basel, Switzerland)
Wang N,Li Y,He F,Liu S,Liu Y,Peng J,Liu J,Yu C,Wang S
Mon Aug 29 00:00:00 UTC 2022
Paeoniflorin directly binds to TNFR1 to regulate podocyte necroptosis in diabetic kidney disease.
Frontiers in pharmacology
Wang X,Liu XQ,Jiang L,Huang YB,Zeng HX,Zhu QJ,Qi XM,Wu YG
Sat Sep 24 00:00:00 UTC 2022
LINC01534/miR-135b-5p/PTPRT axis regulates inflammatory response in loosening total hip replacement via modulating NF-κB signaling pathway.
Injury
Zhang Y,Zhu Q,Fang Q,Yin D,Duan Y,Xue G,Ma N,Cai Y,He M
Wed Jun 01 00:00:00 UTC 2022
Inhibition of cannabinoid receptor type 1 sensitizes triple-negative breast cancer cells to ferroptosis via regulating fatty acid metabolism.
Cell death & disease
Li P,Lin Q,Sun S,Yang N,Xia Y,Cao S,Zhang W,Li Q,Guo H,Zhu M,Wang Y,Zheng Z,Li S
Wed Sep 21 00:00:00 UTC 2022
Synthesis of potent antagonists of receptors for growth hormone-releasing hormone with antitumor and anti-inflammatory activity.
Peptides
Cai R,Zhang X,Wang H,Cui T,Halmos G,Sha W,He J,Popovics P,Vidaurre I,Zhang C,Mirsaeidi M,Schally AV
Fri Apr 01 00:00:00 UTC 2022
Feasibility of Catalpol Intranasal Administration and Its Protective Effect on Acute Cerebral Ischemia in Rats via Anti-Oxidative and Anti-Apoptotic Mechanisms.
Drug design, development and therapy
Wang J,Zhang Y,Zhang M,Sun S,Zhong Y,Han L,Xu Y,Wan D,Zhang J,Zhu H
Mon Mar 28 00:00:00 UTC 2022
Corilagin alleviates intestinal ischemia/reperfusion-induced intestinal and lung injury in mice via inhibiting NLRP3 inflammasome activation and pyroptosis.
Frontiers in pharmacology
Li W,Yang K,Li B,Wang Y,Liu J,Chen D,Diao Y
Wed Dec 21 00:00:00 UTC 2022
Effect of crude polysaccharide from seaweed, Dictyopteris divaricata (CDDP) on gut microbiota restoration and anti-diabetic activity in streptozotocin (STZ)-induced T1DM mice.
Gut pathogens
Siddiqui NZ,Rehman AU,Yousuf W,Khan AI,Farooqui NA,Zang S,Xin Y,Wang L
Sat Sep 17 00:00:00 UTC 2022
Alda-1, an Aldehyde Dehydrogenase 2 Agonist, Improves Cutaneous Wound Healing by Activating Epidermal Keratinocytes via Akt/GSK-3β/β-Catenin Pathway.
Aesthetic plastic surgery
Zhang SM,Yang Z,Qi FZ
Fri Apr 01 00:00:00 UTC 2022
Melanoma stem cells promote metastasis via exosomal miR-1268a inactivation of autophagy.
Biological research
Li X,Liu D,Chen H,Zeng B,Zhao Q,Zhang Y,Chen Y,Wang J,Xing HR
Sat Oct 01 00:00:00 UTC 2022
Involvement of Rab11 in osteoblastic differentiation: Its up-regulation during the differentiation and by tensile stress.
Biochemical and biophysical research communications
Thant L,Kakihara Y,Kaku M,Kitami M,Kitami K,Mizukoshi M,Maeda T,Saito I,Saeki M
Sat Oct 08 00:00:00 UTC 2022
Apelin-13 reduces lipopolysaccharide-induced neuroinflammation and cognitive impairment via promoting glucocorticoid receptor expression and nuclear translocation.
Neuroscience letters
Hu S,Shen P,Chen B,Tian SW,You Y
Sun Sep 25 00:00:00 UTC 2022
Iron-dependent epigenetic modulation promotes pathogenic T cell differentiation in lupus.
The Journal of clinical investigation
Gao X,Song Y,Wu J,Lu S,Min X,Liu L,Hu L,Zheng M,Du P,Yu Y,Long H,Wu H,Jia S,Yu D,Lu Q,Zhao M
Mon May 02 00:00:00 UTC 2022
Effects of Dietary Indole-3-carboxaldehyde Supplementation on Growth Performance, Intestinal Epithelial Function, and Intestinal Microbial Composition in Weaned Piglets.
Frontiers in nutrition
Zhang R,Huang G,Ren Y,Wang H,Ye Y,Guo J,Wang M,Zhu W,Yu K
Sat Jul 16 00:00:00 UTC 2022
Discovery of Potent PROTACs Targeting EGFR Mutants through the Optimization of Covalent EGFR Ligands.
Journal of medicinal chemistry
Zhao HY,Wang HP,Mao YZ,Zhang H,Xin M,Xi XX,Lei H,Mao S,Li DH,Zhang SQ
Thu Mar 24 00:00:00 UTC 2022
Expression Profile and Localization of SARS-CoV-2 Nonstructural Replicase Proteins in Infected Cells.
Microbiology spectrum
Shi FS,Yu Y,Li YL,Cui L,Zhao Z,Wang M,Wang B,Zhang R,Huang YW
Wed Aug 31 00:00:00 UTC 2022
OL-FS13 alleviates experimental cerebral ischemia-reperfusion injury.
Experimental neurology
Liu N,Li Y,Yang Y,Shu L,Liu Y,Wu Y,Sun D,Kang Z,Zhang Y,Ni D,Wei Z,Li S,Yang M,Wang Y,Sun J,Yang X
Tue Nov 01 00:00:00 UTC 2022
Acyl-CoA thioesterase 9 promotes tumour growth and metastasis through reprogramming of fatty acid metabolism in hepatocellular carcinoma.
Liver international : official journal of the International Association for the Study of the Liver
Wang B,Zhang H,Chen YF,Hu LQ,Tian YY,Tong HW,Wang G,Chen C,Yuan P
Tue Nov 01 00:00:00 UTC 2022
The protective effects of butorphanol tartrate against homocysteine-induced blood-brain barrier dysfunction.
Bioengineered
Shen S,Wang J,Zhao Q,Hu Q
Tue Mar 01 00:00:00 UTC 2022
Exploring the pharmacological mechanism of calculus bovis in cerebral ischaemic stroke using a network pharmacology approach.
Journal of ethnopharmacology
Du X,Li C,Zhang S,Sun C,Zhang X,Chen C,Wang X,Cheng F,Wang Q
Thu Feb 10 00:00:00 UTC 2022
Autophagy Mediates Escherichia Coli-Induced Cellular Inflammatory Injury by Regulating Calcium Mobilization, Mitochondrial Dysfunction, and Endoplasmic Reticulum Stress.
International journal of molecular sciences
Liu J,Qiu R,Liu R,Song P,Lin P,Chen H,Zhou D,Wang A,Jin Y
Wed Nov 16 00:00:00 UTC 2022
DSF/Cu induces antitumor effect against diffuse large B-cell lymphoma through suppressing NF-κB/BCL6 pathways.
Cancer cell international
Zhu Y,Lei C,Jiang Q,Yu Q,Qiu L
Tue Jul 26 00:00:00 UTC 2022
Shikonin induces apoptosis and autophagy via downregulation of pyrroline-5-carboxylate reductase1 in hepatocellular carcinoma cells.
Bioengineered
Zhang J,Shang L,Jiang W,Wu W
Tue Mar 01 00:00:00 UTC 2022
Anp32e promotes renal interstitial fibrosis by upregulating the expression of fibrosis-related proteins.
International journal of biological sciences
Wei J,Shan Y,Xiao Z,Wen L,Tao Y,Fang X,Luo H,Tang C,Li Y
Mon Oct 24 00:00:00 UTC 2022
Inhibition of the proliferation, invasion, migration, and epithelial-mesenchymal transition of prostate cancer cells through the action of ATP1A2 on the TGF-β/Smad pathway.
Translational andrology and urology
Zhang B,Zhu Z,Zhang X,Li F,Ding A
Sat Jan 01 00:00:00 UTC 2022
Characterization of two novel knock-in mouse models of syndromic retinal ciliopathy carrying hypomorphic Sdccag8 mutations.
Zoological research
Ren ZL,Zhang HB,Li L,Yang ZL,Jiang L
Wed May 18 00:00:00 UTC 2022
Triad3A-Dependent TLR4 Ubiquitination and Degradation Contributes to the Anti-Inflammatory Effects of Pterostilbene on Vascular Dementia.
Journal of agricultural and food chemistry
Xu J,Liu J,Mi Y,Zhao T,Mu D,Meng Q,Wang F,Li N,Hou Y
Wed May 18 00:00:00 UTC 2022
Capsaicin suppresses hepatocarcinogenesis by inhibiting the stemness of hepatic progenitor cells via SIRT1/SOX2 signaling pathway.
Cancer medicine
Xie ZQ,Li HX,Hou XJ,Huang MY,Zhu ZM,Wei LX,Tang CX
Tue Nov 01 00:00:00 UTC 2022
In vitro and in silico anti-osteoporosis activities and underlying mechanisms of a fructan, ABW90-1, from Achyranthes bidentate.
Carbohydrate polymers
Li T,Hou X,Huang Y,Wang C,Chen H,Yan C
Sat Jan 15 00:00:00 UTC 2022
Eurycomanol alleviates hyperuricemia by promoting uric acid excretion and reducing purine synthesis.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Bao R,Chen Q,Li Z,Wang D,Wu Y,Liu M,Zhang Y,Wang T
Tue Feb 01 00:00:00 UTC 2022
Long noncoding RNA GAS5 inhibits proliferation and metastasis in papillary thyroid carcinoma through the IFN/STAT1 signaling pathway.
Pathology, research and practice
Xu Y,Lu J,Lou N,Lu W,Xu J,Jiang H,Ye G
Sun May 01 00:00:00 UTC 2022
Silencing of Rab23 by siRNA inhibits ultraviolet B-induced melanogenesis via downregulation of PKA/CREB/MITF.
Experimental dermatology
Wei X,Huang M,Yang Y,Liu Y,Chi S,Li C
Mon Aug 01 00:00:00 UTC 2022
The single-atom iron nanozyme mimicking peroxidase remodels energy metabolism and tumor immune landscape for synergistic chemodynamic therapy and photothermal therapy of triple-negative breast cancer.
Frontiers in bioengineering and biotechnology
Qian X,Shi R,Chen J,Wang Y,Han X,Sun Y,Ling C,Wang G,Xu AW,Pan Y
Sat Nov 19 00:00:00 UTC 2022
Inhibition of human peptide deformylase by actinonin sensitizes glioblastoma cells to temozolomide chemotherapy.
Experimental cell research
Lan B,Zhao H,He Y,Zhao Z,Wang N,Gao Y
Tue Nov 15 00:00:00 UTC 2022
HMGB2 causes photoreceptor death via down-regulating Nrf2/HO-1 and up-regulating NF-κB/NLRP3 signaling pathways in light-induced retinal degeneration model.
Free radical biology & medicine
Zhang Y,Zhao Z,Zhao X,Xie H,Zhang C,Sun X,Zhang J
Tue Mar 01 00:00:00 UTC 2022
RBMS2 Chemosensitizes Breast Cancer Cells to Doxorubicin by Regulating BMF Expression.
International journal of biological sciences
Xu F,Xia T,Xu QT,Zhang X,Huang YZ,Sun X,Shi L,Zhou XJ,Wei JF,Ding Q
Fri Apr 15 00:00:00 UTC 2022
Inhibitory Effects of Parachlorella Beijerinckii Extracts on the Formation of Advanced Glycation End Products and Glycative Stress-Induced Inflammation in an In Vitro Skin Dermis-Like Model.
Evidence-based complementary and alternative medicine : eCAM
Imai Y,Nakashima Y,Kanno T
Fri Nov 18 00:00:00 UTC 2022
Integrated Strategies of Diverse Feature Selection Methods Identify Aging-Based Reliable Gene Signatures for Ischemic Cardiomyopathy.
Frontiers in molecular biosciences
Song H,Chen S,Zhang T,Huang X,Zhang Q,Li C,Chen C,Chen S,Liu D,Wang J,Tu Y,Wu Y,Liu Y
Sat Mar 19 00:00:00 UTC 2022
The Effects of Hedgehog Signaling Pathway on the Proliferation and Apoptosis of Melanoma Cells.
Journal of oncology
Peng ZP,Huang SF,Li JJ,Tang XK,Wang XY,Li HM
Sat Apr 30 00:00:00 UTC 2022
Sodium Butyrate Attenuated Diabetes-Induced Intestinal Inflammation by Modulating Gut Microbiota.
Evidence-based complementary and alternative medicine : eCAM
Liu L,Chen Y,Wu Q,Shu A,Sun J
Wed Sep 07 00:00:00 UTC 2022
Rosa centifolia petal extract induces endothelium-dependent and endothelium-independent vasorelaxation in rat aorta and prevents accumulation of inflammatory factors in human umbilical vein endothelial cells.
Journal of food biochemistry
Kaneda H,Hori M,Shinomiya H,Nakajima A,Yamazaki S,Sasaki N,Sato T,Kaneda T
Fri Jul 01 00:00:00 UTC 2022
Menadione Potentiates Auranofin-Induced Glioblastoma Cell Death.
International journal of molecular sciences
Szeliga M,Rola R
Sun Dec 11 00:00:00 UTC 2022
Increased expression of osteopontin in subchondral bone promotes bone turnover and remodeling, and accelerates the progression of OA in a mouse model.
Aging
Lin C,Chen Z,Guo D,Zhou L,Lin S,Li C,Li S,Wang X,Lin B,Ding Y
Tue Jan 04 00:00:00 UTC 2022
Dual-fluorescent reporter for live-cell imaging of the ER during DENV infection.
Frontiers in cellular and infection microbiology
Corliss L,Holliday M,Lennemann NJ
Mon Nov 21 00:00:00 UTC 2022
Down-Regulation of Double C2 Domain Alpha Promotes the Formation of Hyperplastic Nerve Fibers in Aganglionic Segments of Hirschsprung's Disease.
International journal of molecular sciences
Xiao J,Meng X,Chen K,Wang J,Wu L,Chen Y,Yu X,Feng J,Li Z
Tue Sep 06 00:00:00 UTC 2022
Autophagy and mitochondrial dynamics contribute to the protective effect of diosgenin against 3-MCPD induced kidney injury.
Chemico-biological interactions
Zhong Y,Sun D,Yao Y,Liu Q,Guo T,Wang X,Peng X
Tue Mar 01 00:00:00 UTC 2022
DNA Methyltransferase Inhibitor 5-AZA-DC Regulates TGFβ1-Mediated Alteration of Neuroglial Cell Functions after Oxidative Stress.
Oxidative medicine and cellular longevity
Zhao XY,Zhang XL
Tue Oct 11 00:00:00 UTC 2022
Ciclopirox Olamine Exerts Tumor-Suppressor Effects via Topoisomerase II Alpha in Lung Adenocarcinoma.
Frontiers in oncology
Yin J,Che G,Jiang K,Zhou Z,Wu L,Xu M,Liu J,Yan S
Mon May 02 00:00:00 UTC 2022
Munronoid I Ameliorates DSS-Induced Mouse Colitis by Inhibiting NLRP3 Inflammasome Activation and Pyroptosis Via Modulation of NLRP3.
Frontiers in immunology
Ma X,Di Q,Li X,Zhao X,Zhang R,Xiao Y,Li X,Wu H,Tang H,Quan J,Wu Z,Xiao W,Chen W
Mon Jul 25 00:00:00 UTC 2022
Phyllanthus Niruri L. Exerts Protective Effects Against the Calcium Oxalate-Induced Renal Injury via Ellgic Acid.
Frontiers in pharmacology
Li MT,Liu LL,Zhou Q,Huang LX,Shi YX,Hou JB,Lu HT,Yu B,Chen W,Guo ZY
Mon Jun 26 00:00:00 UTC 2023
Preparation, Characterization, Evaluation of Neuroprotective Effect, and Related Mechanisms of Phosphatidylserine Emulsion in 5- and 12-Week Old Mice.
Journal of agricultural and food chemistry
Liu B,Zhang N,Yang J,Sun W,Zhang R,Zheng X,Wang Z,Siebert HC,Han J
Wed Feb 16 00:00:00 UTC 2022
Effects of 4-vinylcyclohexene diepoxide on the cell cycle, apoptosis, and steroid hormone secretion of goat ovarian granulosa cells.
In vitro cellular & developmental biology. Animal
Miao Y,Wan W,Zhu K,Pan M,Zhao X,Ma B,Wei Q
Tue Mar 01 00:00:00 UTC 2022
Reduction of H3K27cr Modification During DNA Damage in Colon Cancer.
Frontiers in oncology
Liao M,Chu W,Sun X,Zheng W,Gao S,Li D,Pei D
Tue Aug 09 00:00:00 UTC 2022
Spermidine protects against acute kidney injury by modulating macrophage NLRP3 inflammasome activation and mitochondrial respiration in an eIF5A hypusination-related pathway.
Molecular medicine (Cambridge, Mass.)
Li X,Zhou X,Liu X,Li X,Jiang X,Shi B,Wang S
Sun Sep 04 00:00:00 UTC 2022
SREBP2/Rab11s/GLUT1/6 network regulates proliferation and migration of glioblastoma.
Pathology, research and practice
Cheng C,Tu J,Hu Z,Chen Y,Wang Y,Zhang T,Zhang C,Li C,Wang Y,Niu C
Thu Dec 01 00:00:00 UTC 2022
miR-153-3p inhibits osteogenic differentiation of BMSCs by down-regulating the expression of RUNX2 in a high glucose environment.
American journal of translational research
Wang Q,Xia Q,Meng M,Li Y,Tang Z,Zeng X,Chen H,Shu J,Xv X,Chen J,Lu J,Wang H,Ye Z,Song B,Dong Q
Sat Nov 19 00:00:00 UTC 2022
Cellular PSMB4 Protein Suppresses Influenza A Virus Replication through Targeting NS1 Protein.
Viruses
Yang CH,Hsu CF,Lai XQ,Chan YR,Li HC,Lo SY
Mon Oct 17 00:00:00 UTC 2022
Therapeutic targeting m6A-guided miR-146a-5p signaling contributes to the melittin-induced selective suppression of bladder cancer.
Cancer letters
Yan R,Dai W,Wu R,Huang H,Shu M
Sat May 28 00:00:00 UTC 2022
The involvement of TRPM2 on the MPP+-induced oxidative neurotoxicity and apoptosis in hippocampal neurons from neonatal mice: protective role of resveratrol.
Neurological research
Yildizhan K,Çinar R,Naziroğlu M
Fri Jul 01 00:00:00 UTC 2022
Xiao-Chai-Hu Decoction Ameliorates Poly (I:C)-Induced Viral Pneumonia through Inhibiting Inflammatory Response and Modulating Serum Metabolism.
Evidence-based complementary and alternative medicine : eCAM
Chen F,Qu F,Jia Y,Wang C,Yu Y,Liao J,Lin M,Chen F,Sun Z
Sat Jul 23 00:00:00 UTC 2022
Interferon-Stimulated Gene 15 Knockout in Mice Impairs IFNα-Mediated Antiviral Activity.
Viruses
Li C,He WF,Li LX,Chen J,Yang GQ,Chang HT,Liu HM
Wed Aug 24 00:00:00 UTC 2022
Consistent downregulation of the cleft lip/palate-associated genes IRF6 and GRHL3 in carcinomas.
Frontiers in oncology
Parisi L,Mockenhaupt C,Rihs S,Mansour F,Katsaros C,Degen M
Sat Dec 03 00:00:00 UTC 2022
Suffruticosol C-Mediated Autophagy and Cell Cycle Arrest via Inhibition of mTORC1 Signaling.
Nutrients
Qin S,Geng H,Wang G,Chen L,Xia C,Yao J,Bai Z,Deng L
Thu Nov 24 00:00:00 UTC 2022
Heat Stress Modulates a Placental Immune Response Associated With Alterations in the Development of the Fetal Intestine and Its Innate Immune System in Late Pregnant Mouse.
Frontiers in physiology
Guo H,Liu R,He J,Yao W,Zheng W
Sat Jul 16 00:00:00 UTC 2022
Chronic constant light exposure aggravates high fat diet-induced renal injury in rats.
Frontiers in endocrinology
Xing L,Wu S,Shi Y,Yue F,Wei L,Russell R,Zhang D
Tue Aug 16 00:00:00 UTC 2022
Myo1b promotes tumor progression and angiogenesis by inhibiting autophagic degradation of HIF-1α in colorectal cancer.
Cell death & disease
Chen YH,Xu NZ,Hong C,Li WQ,Zhang YQ,Yu XY,Huang YL,Zhou JY
Tue Nov 08 00:00:00 UTC 2022
Renoprotective effects of oleanolic acid and its possible mechanisms in rats with diabetic kidney disease.
Biochemical and biophysical research communications
Liu Y,Hu Z,Xing H,Kang L,Chen X,Liu B,Niu K
Sun Dec 25 00:00:00 UTC 2022
DR1 activation promotes vascular smooth muscle cell apoptosis via up-regulation of CSE/H2 S pathway in diabetic mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wen X,Xi Y,Zhang Y,Jiao L,Shi S,Bai S,Sun F,Chang G,Wu R,Hao J,Li H
Sat Jan 01 00:00:00 UTC 2022
Porcine Enteric Coronavirus PEDV Induces the ROS-ATM and Caspase7-CAD-γH2AX Signaling Pathways to Foster Its Replication.
Viruses
Ming X,Chen H,Yang Y,Zhao P,Sun L,Zhang C,Shin HJ,Lee JS,Jung YS,Qian Y
Mon Aug 15 00:00:00 UTC 2022
Dehydrocostus Lactone Suppresses Dextran Sulfate Sodium-Induced Colitis by Targeting the IKKα/β-NF-κB and Keap1-Nrf2 Signalling Pathways.
Frontiers in pharmacology
Yuan Y,Hu Q,Liu L,Xie F,Yang L,Li Y,Zhang C,Chen H,Tang J,Shen X
Fri Mar 25 00:00:00 UTC 2022
Pirh2 restricts influenza A virus replication by modulating short-chain ubiquitination of its nucleoprotein.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Chen H,Gao X,Zhao S,Bao C,Ming X,Qian Y,Zhou Y,Jung YS
Sat Oct 01 00:00:00 UTC 2022
A critical ETV4/Twist1/Vimentin axis in Ha-RAS-induced aggressive breast cancer.
Cancer gene therapy
Liu W,Gajendran B,Sample KM,Wang C,Hu A,Chen B,Li Y,Zacksenhaus E,Ben-David Y
Tue Nov 01 00:00:00 UTC 2022
Substitution of the SERCA2 Cys674 reactive thiol accelerates atherosclerosis by inducing endoplasmic reticulum stress and inflammation.
British journal of pharmacology
Su H,Mei Y,Luo S,Wu H,He Y,Shiraishi Y,Hu P,Cohen RA,Tong X
Sat Oct 01 00:00:00 UTC 2022
Hyperoside protects against cyclophosphamide induced ovarian damage and reduced fertility by suppressing HIF-1α/BNIP3-mediated autophagy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhu F,Gao J,Zeng F,Lai Y,Ruan X,Deng G
Thu Dec 01 00:00:00 UTC 2022
Ferulic acid attenuates high glucose-induced apoptosis in retinal pigment epithelium cells and protects retina in db/db mice.
PeerJ
Zhu D,Zou W,Cao X,Xu W,Lu Z,Zhu Y,Hu X,Hu J,Zhu Q
Thu Jan 12 00:00:00 UTC 2023
MCTS1 promotes laryngeal squamous cell carcinoma cell growth via enhancing LARP7 stability.
Clinical and experimental pharmacology & physiology
Yang M,Ma B,Liu X
Wed Jun 01 00:00:00 UTC 2022
Facilitating Mitophagy via Pink1/Parkin2 Signaling Is Essential for the Neuroprotective Effect of β-Caryophyllene against CIR-Induced Neuronal Injury.
Brain sciences
Rao J,Wu Y,Fan X,Yang S,Jiang L,Dong Z,Chen S
Thu Jun 30 00:00:00 UTC 2022
Identification of endocrine-disrupting chemicals targeting the genes and pathways of genital anomalies in males.
Ecotoxicology and environmental safety
Zhou X,Zhang X,Zhou X,Abulimiti G,Wang Y,Zhang Q,Cong R,Ji C,Luan J,Yao L,Yang J,Song N
Thu Dec 01 00:00:00 UTC 2022
LINC00941 promotes pancreatic cancer malignancy by interacting with ANXA2 and suppressing NEDD4L-mediated degradation of ANXA2.
Cell death & disease
Wang J,He Z,Liu X,Xu J,Jiang X,Quan G,Jiang J
Thu Aug 18 00:00:00 UTC 2022
Apelin-13 Attenuates Lipopolysaccharide-Induced Inflammatory Responses and Acute Lung Injury by Regulating PFKFB3-Driven Glycolysis Induced by NOX4-Dependent ROS.
Journal of inflammation research
Yuan Y,Wang W,Zhang Y,Hong Q,Huang W,Li L,Xie Z,Chen Y,Li X,Meng Y
Fri Apr 08 00:00:00 UTC 2022
SorCS3 promotes the internalization of p75NTR to inhibit GBM progression.
Cell death & disease
Zhang Y,Li Y,Fan Y,Zhang X,Tang Z,Qi J,Zhao B,Li F,Chen X,Liang H,Xu H,Li D
Thu Apr 07 00:00:00 UTC 2022
Cannabis suppresses antitumor immunity by inhibiting JAK/STAT signaling in T cells through CNR2.
Signal transduction and targeted therapy
Xiong X,Chen S,Shen J,You H,Yang H,Yan C,Fang Z,Zhang J,Cai X,Dong X,Kang T,Li W,Zhou P
Wed Apr 06 00:00:00 UTC 2022
Dihydroartemisinin attenuates osteoclast formation and bone resorption via inhibiting the NF‑κB, MAPK and NFATc1 signaling pathways and alleviates osteoarthritis.
International journal of molecular medicine
Ding D,Yan J,Feng G,Zhou Y,Ma L,Jin Q
Sat Jan 01 00:00:00 UTC 2022
miR-141-3p affects β-catenin signaling and apoptosis by targeting Ubtd2 in rats with anorectal malformations.
Annals of the New York Academy of Sciences
Wang CY,Li SY,Xiao YX,Zhen L,Wei XG,Tang XB,Yuan ZW,Bai YZ
Thu Dec 01 00:00:00 UTC 2022
Liensinine Inhibits Cell Growth and Blocks Autophagic Flux in Nonsmall-Cell Lung Cancer.
Journal of oncology
Chang M,Ding S,Dong X,Shang X,Li Y,Xie L,Song X,Song X
Sat Jul 16 00:00:00 UTC 2022
Tooth Loss Induces Memory Impairment and Glial Activation in Young Wild-Type Mice.
Journal of Alzheimer's disease reports
Taslima F,Abdelhamid M,Zhou C,Chen Y,Jung CG,Michikawa M
Wed Dec 21 00:00:00 UTC 2022
Analgesic effect of nobiletin against neuropathic pain induced by the chronic constriction injury of the sciatic nerve in mice.
Phytotherapy research : PTR
Zhu C,Tian M,Liu N,Ma L,Lan X,Yang J,Du J,Ma H,Li Y,Zheng P,Yu J,Peng X
Thu Sep 01 00:00:00 UTC 2022
Effects of butein on human osteosarcoma cell proliferation, apoptosis, and autophagy through oxidative stress.
Human & experimental toxicology
Zhang P,Zhang J,Quan H,Chen P,Wang J,Liang Y
Wed Mar 02 00:00:00 UTC 2022
Nrf2 improves hippocampal synaptic plasticity, learning and memory through the circ-Vps41/miR-26a-5p/CaMKIV regulatory network.
Experimental neurology
Zhang R,Gao Y,Li Y,Geng D,Liang Y,He Q,Wang L,Cui H
Sun May 01 00:00:00 UTC 2022
Genetic Deletion of HLJ1 Does Not Affect Blood Coagulation in Mice.
International journal of molecular sciences
Hsu MC,Luo WJ,Guo BC,Chen CH,Hu PA,Tsai YH,Su KY,Lee TS
Sun Feb 13 00:00:00 UTC 2022
Histone Deacetylase Inhibitor Panobinostat Benefits the Therapeutic Efficacy of Oncolytic Herpes Simplex Virus Combined with PD-1/PD-L1 Blocking in Glioma and Squamous Cell Carcinoma Models.
Viruses
Wu Y,Chen X,Wang L,Zhou X,Liu Y,Ji D,Ren P,Zhou GG,Zhao J
Thu Dec 15 00:00:00 UTC 2022
Hypoxia-Induced miR-210 Promotes Endothelial Cell Permeability and Angiogenesis via Exosomes in Pancreatic Ductal Adenocarcinoma.
Biochemistry research international
Wu G,Ding X,Quan G,Xiong J,Li Q,Li Z,Wang Y
Tue Dec 06 00:00:00 UTC 2022
CD147 Mediates 5-Fluorouracil Resistance in Colorectal Cancer by Reprogramming Glycolipid Metabolism.
Frontiers in oncology
Dong S,Li S,Wang X,Liang S,Zhang W,Li L,Xu Q,Shi B,Cheng Z,Zhang X,Zhong M,Zhang G,Hu S
Fri Jul 29 00:00:00 UTC 2022
Evaluation of decompressive craniectomy in mice after severe traumatic brain injury.
Frontiers in neurology
Liu Y,Liu X,Chen Z,Wang Y,Li J,Gong J,He A,Zhao M,Yang C,Yang W,Wang Z
Sat Aug 13 00:00:00 UTC 2022
Involvement of hedgehog signaling in all-trans retinoic acid-mediated suppression of colon cancer.
American journal of translational research
Xu Y,Sun H
Wed Oct 19 00:00:00 UTC 2022
MiR-145 inhibits the differentiation and proliferation of bone marrow stromal mesenchymal stem cells by GABARAPL1 in steroid-induced femoral head necrosis.
BMC musculoskeletal disorders
Xu P,Chang J,Ma G,Liao F,Xu T,Wu Y,Yin Z
Sat Nov 26 00:00:00 UTC 2022
Mitotic SENP3 activation couples with cGAS signaling in tumor cells to stimulate anti-tumor immunity.
Cell death & disease
Hu G,Chen Y,Yang X,Wang Y,He J,Wang T,Fan Q,Deng L,Tu J,Tan H,Cheng J
Fri Jul 22 00:00:00 UTC 2022
Ventilator-induced lung injury results in oxidative stress response and mitochondrial swelling in a mouse model.
Laboratory animal research
Joelsson JP,Asbjarnarson A,Sigurdsson S,Kricker J,Valdimarsdottir B,Thorarinsdottir H,Starradottir E,Gudjonsson T,Ingthorsson S,Karason S
Fri Jul 22 00:00:00 UTC 2022
Reticulon 2 promotes gastric cancer metastasis via activating endoplasmic reticulum Ca2+ efflux-mediated ERK signalling.
Cell death & disease
Song S,Liu B,Zeng X,Wu Y,Chen H,Wu H,Gu J,Gao X,Ruan Y,Wang H
Fri Apr 15 00:00:00 UTC 2022
Silencing CAMK2D Promotes the Proliferation of Spermatogonia in the Testis of Experimental Varicocele Rats.
Evidence-based complementary and alternative medicine : eCAM
Tang S,Yang P,Ding Y,Chen Q,Huang H,Chen X,Zhou H
Fri Aug 18 00:00:00 UTC 2023
Therapeutic Potential of Synthetic Human β-Defensin 1 Short Motif Pep-B on Lipopolysaccharide-Stimulated Human Dental Pulp Stem Cells.
Mediators of inflammation
Shi J,Hu Z,Zhou Y,Zuo M,Wu H,Jin W,Xie Z
Thu Mar 31 00:00:00 UTC 2022
Osteoclast-Driven Osteogenesis, Bone Remodeling and Biomaterial Resorption: A New Profile of BMP2-CPC-Induced Alveolar Bone Regeneration.
International journal of molecular sciences
Shen H,Zhuang Y,Zhang C,Zhang C,Yuan Y,Yu H,Si J,Shen G
Thu Oct 13 00:00:00 UTC 2022
Nintedanib Inhibits Endothelial Mesenchymal Transition in Bleomycin-Induced Pulmonary Fibrosis via Focal Adhesion Kinase Activity Reduction.
International journal of molecular sciences
Yu WK,Chen WC,Su VY,Shen HC,Wu HH,Chen H,Yang KY
Mon Jul 25 00:00:00 UTC 2022
EspF of Enterohemorrhagic Escherichia coli Enhances Apoptosis via Endoplasmic Reticulum Stress in Intestinal Epithelial Cells: An Isobaric Tags for Relative and Absolute Quantitation-Based Comparative Proteomic Analysis.
Frontiers in microbiology
Wang X,Yan K,Fu M,Liang S,Zhao H,Fu C,Yang L,Song Z,Sun D,Wan C
Tue Jul 19 00:00:00 UTC 2022
Long noncoding RNA H19 mediates neural stem/progenitor cells proliferation, differentiation and apoptosis through the p53 signaling pathway after ischemic stroke.
Biochemical and biophysical research communications
Gan L,Liao S,Tong Y,Li W,Peng W,Deng S
Tue Mar 15 00:00:00 UTC 2022
OAS1, OAS2, and OAS3 Contribute to Epidermal Keratinocyte Proliferation by Regulating Cell Cycle and Augmenting IFN-1‒Induced Jak1‒Signal Transducer and Activator of Transcription 1 Phosphorylation in Psoriasis.
The Journal of investigative dermatology
Huang YZ,Zheng YX,Zhou Y,Xu F,Cui YZ,Chen XY,Wang ZY,Yan BX,Zheng M,Man XY
Sat Oct 01 00:00:00 UTC 2022
Identification of HMGCR as the anticancer target of physapubenolide against melanoma cells by in silico target prediction.
Acta pharmacologica Sinica
Wang HY,Yu P,Chen XS,Wei H,Cao SJ,Zhang M,Zhang Y,Tao YG,Cao DS,Qiu F,Cheng Y
Wed Jun 01 00:00:00 UTC 2022
Neutrophils Promote Tumor Progression in Oral Squamous Cell Carcinoma by Regulating EMT and JAK2/STAT3 Signaling Through Chemerin.
Frontiers in oncology
Hu X,Xiang F,Feng Y,Gao F,Ge S,Wang C,Zhang X,Wang N
Wed Feb 16 00:00:00 UTC 2022
A 3D-printed molybdenum-containing scaffold exerts dual pro-osteogenic and anti-osteoclastogenic effects to facilitate alveolar bone repair.
International journal of oral science
Tian B,Li X,Zhang J,Zhang M,Gan D,Deng D,Sun L,He X,Wu C,Chen F
Mon Sep 05 00:00:00 UTC 2022
Acetyl-Coenzyme A Synthetase 2 Potentiates Macropinocytosis and Muscle Wasting Through Metabolic Reprogramming in Pancreatic Cancer.
Gastroenterology
Zhou Z,Ren Y,Yang J,Liu M,Shi X,Luo W,Fung KM,Xu C,Bronze MS,Zhang Y,Houchen CW,Li M
Tue Nov 01 00:00:00 UTC 2022
A novel regulatory mechanism of geniposide for improving glucose homeostasis mediated by circulating RBP4.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu J,Song C,Nie C,Sun Y,Wang Y,Xue L,Fan M,Qian H,Wang L,Li Y
Sat Jan 01 00:00:00 UTC 2022
Irisin attenuates P. gingivalis-suppressed osteogenic/cementogenic differentiation of periodontal ligament cells via p38 signaling pathway.
Biochemical and biophysical research communications
Huang X,Xiao J,Wang X,Cao Z
Sat Aug 27 00:00:00 UTC 2022
The Effects and Mechanisms of the Rapamycin-eluting Stent in Urethral Stricture Prevention in Rabbits.
Balkan medical journal
Zhang T,Zhao W,Ren T,Chen J,Chen Z,Wang Y,Cheng X,Wu J,Yuan C,He T
Mon Mar 14 00:00:00 UTC 2022
Psychosocial stress and cannabinoid drugs affect acetylation of α-tubulin (K40) and gene expression in the prefrontal cortex of adult mice.
PloS one
Tomas-Roig J,Ramasamy S,Zbarsky D,Havemann-Reinecke U,Hoyer-Fender S
Fri Sep 23 00:00:00 UTC 2022
The role of AMPK-Sirt1-autophagy pathway in the intestinal protection process by propofol against regional ischemia/reperfusion injury in rats.
International immunopharmacology
Liu X,Yang B,Tan YF,Feng JG,Jia J,Yang CJ,Chen Y,Wang MH,Zhou J
Sat Oct 01 00:00:00 UTC 2022
Retracted Article: LncRNA KCNQ1OT1 Promotes Apoptosis and Oxidative Stress of Human Lens Epithelial Cells through Epigenetic Regulation of WRN.
Current eye research
Jiang S,Jia Y,Gao Z
Wed Jun 01 00:00:00 UTC 2022
Single Treatment of Vitamin D3 Ameliorates LPS-Induced Acute Lung Injury through Changing Lung Rodentibacter abundance.
Molecular nutrition & food research
Jin A,Zhao Y,Yuan Y,Ma S,Chen J,Yang X,Lu S,Sun Q
Tue Feb 01 00:00:00 UTC 2022
Bioelectric fields coordinate wound contraction and re-epithelialization process to accelerate wound healing via promoting myofibroblast transformation.
Bioelectrochemistry (Amsterdam, Netherlands)
Yang J,Liu X,Wang W,Chen Y,Liu J,Zhang Z,Wu C,Jiang X,Liang Y,Zhang J
Thu Dec 01 00:00:00 UTC 2022
α-Terthienyl induces prostate cancer cell death through inhibiting androgen receptor expression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gan X,Huang H,Wen J,Liu K,Yang Y,Li X,Fang G,Liu Y,Wang X
Mon Aug 01 00:00:00 UTC 2022
The comprehensive expression and functional analysis of m6A modification "readers" in hepatocellular carcinoma.
Aging
Qin S,Liu G,Jin H,Chen X,He J,Xiao J,Qin Y,Mao Y,Zhao L
Fri Aug 12 00:00:00 UTC 2022
CD36 aggravates podocyte injury by activating NLRP3 inflammasome and inhibiting autophagy in lupus nephritis.
Cell death & disease
Lv F,He Y,Xu H,Li Y,Han L,Yan L,Lang H,Zhao Y,Zhao Z,Qi Y
Tue Aug 23 00:00:00 UTC 2022
Transient and Prolonged Activation of Wnt Signaling Contribute Oppositely to the Pathogenesis of Asherman's Syndrome.
International journal of molecular sciences
Xue X,Li X,Yao J,Zhang X,Ren X,Xu S
Mon Aug 08 00:00:00 UTC 2022
Identifying potential prognosis markers in hepatocellular carcinoma via integrated bioinformatics analysis and biological experiments.
Frontiers in genetics
Hu X,Zhou J,Zhang Y,Zeng Y,Jie G,Wang S,Yang A,Zhang M
Sat Aug 06 00:00:00 UTC 2022
Exosomal circGSE1 promotes immune escape of hepatocellular carcinoma by inducing the expansion of regulatory T cells.
Cancer science
Huang M,Huang X,Huang N
Wed Jun 01 00:00:00 UTC 2022
Pectin supplementation ameliorates intestinal epithelial barrier function damage by modulating intestinal microbiota in lipopolysaccharide-challenged piglets.
The Journal of nutritional biochemistry
Wen X,Zhong R,Dang G,Xia B,Wu W,Tang S,Tang L,Liu L,Liu Z,Chen L,Zhang H
Tue Nov 01 00:00:00 UTC 2022
Resveratrol pretreatment protects neurons from oxygen-glucose deprivation/reoxygenation and ischemic injury through inhibiting ferroptosis.
Bioscience, biotechnology, and biochemistry
Zhu H,Huang J,Chen Y,Li X,Wen J,Tian M,Ren J,Zhou L,Yang Q
Tue May 24 00:00:00 UTC 2022
Mild hypothermia ameliorates hepatic ischemia reperfusion injury by inducing RBM3 expression.
Apoptosis : an international journal on programmed cell death
Xiao Q,Liu Y,Zhang X,Liu Z,Xiao J,Ye Q,Fu B
Thu Dec 01 00:00:00 UTC 2022
Cognitive-exercise dual-task intervention ameliorates cognitive decline in natural aging rats through reducing oxidative stress and enhancing synaptic plasticity.
Experimental gerontology
Li XL,Tao X,Li TC,Zhu ZM,Huang PL,Gong WJ
Tue Nov 01 00:00:00 UTC 2022
Concomitant pyroptotic and apoptotic cell death triggered in macrophages infected by Zika virus.
PloS one
Wen C,Yu Y,Gao C,Qi X,Cardona CJ,Xing Z
Mon Apr 25 00:00:00 UTC 2022
BMP-7 ameliorates partial epithelial-mesenchymal transition by restoring SnoN protein level via Smad1/5 pathway in diabetic kidney disease.
Cell death & disease
Peng W,Zhou X,Xu T,Mao Y,Zhang X,Liu H,Liang L,Liu L,Liu L,Xiao Y,Zhang F,Li S,Shi M,Zhou Y,Tang L,Wang Y,Guo B
Mon Mar 21 00:00:00 UTC 2022
Fucoidan ameliorates glucose metabolism by the improvement of intestinal barrier and inflammatory damage in type 2 diabetic rats.
International journal of biological macromolecules
Liu Y,Xu Z,Huang H,Xue Y,Zhang D,Zhang Y,Li W,Li X
Tue Mar 15 00:00:00 UTC 2022
Differential expression pattern, bioinformatics analysis, and validation of circRNA and mRNA in patients with arteriosclerosis.
Frontiers in cardiovascular medicine
Liu Y,Wang K,Li G,Chen Z
Sat Oct 01 00:00:00 UTC 2022
Fucoidan protects the pancreas and improves glucose metabolism through inhibiting inflammation and endoplasmic reticulum stress in T2DM rats.
Food & function
Liu Y,Huang H,Xu Z,Xue Y,Zhang D,Zhang Y,Li W,Li X
Mon Mar 07 00:00:00 UTC 2022
TiO2 nanosheets promote the transformation of vascular smooth muscle cells into foam cells in vitro and in vivo through the up-regulation of nuclear factor kappa B subunit 2.
Journal of hazardous materials
Li S,Yan D,Huang C,Yang F,Cao Y
Tue Feb 15 00:00:00 UTC 2022
The neural stem cell properties of Pkd2l1+ cerebrospinal fluid-contacting neurons in vivo.
Frontiers in cellular neuroscience
Cao L,Huang MZ,Zhang Q,Luo ZR,Zhang Y,An PJ,Yang LL,Tan W,Wang CQ,Dou XW,Li Q
Wed Sep 28 00:00:00 UTC 2022
Placenta-Targeted Nanoparticles Loaded with PFKFB3 Overexpression Plasmids Enhance Angiogenesis and Placental Function.
Bioengineering (Basel, Switzerland)
Li Q,Liu X,Liu W,Zhang Y,Liu W,Wu M,Chen Z,Zhao Y,Zou L
Fri Nov 04 00:00:00 UTC 2022
Integrated multi-omics analysis reveals miR-20a as a regulator for metabolic colorectal cancer.
Heliyon
Song K,Liu C,Zhang J,Yao Y,Xiao H,Yuan R,Li K,Yang J,Zhao W,Zhang Y
Tue Mar 01 00:00:00 UTC 2022
O-GlcNAc modification mediates aquaporin 3 to coordinate endometrial cell glycolysis and affects embryo implantation.
Journal of advanced research
Zhang H,Qi J,Pei J,Zhang M,Shang Y,Li Z,Wang Y,Guo J,Sun K,Fan J,Sui L,Xu Y,Kong L,Kong Y
Tue Mar 01 00:00:00 UTC 2022
Effect of Astragalus polysaccharides on the cryopreservation of goat semen.
Theriogenology
Zhang X,Hu ZT,Li Y,Li YX,Xian M,Guo SM,Hu JH
Tue Nov 01 00:00:00 UTC 2022
Dihydroartemisinin represses oral squamous cell carcinoma progression through downregulating mitochondrial calcium uniporter.
Bioengineered
Zheng S,Wu R,Deng Y,Zhang Q
Sat Jan 01 00:00:00 UTC 2022
EGCG Enhances the Chemosensitivity of Colorectal Cancer to Irinotecan through GRP78-MediatedEndoplasmic Reticulum Stress.
Journal of oncology
Wu W,Gou H,Xiang B,Geng R,Dong J,Yang X,Chen D,Dai R,Chen L,Liu J
Sat Sep 24 00:00:00 UTC 2022
β-Carotene Attenuates Apoptosis and Autophagy via PI3K/AKT/mTOR Signaling Pathway in Necrotizing Enterocolitis Model Cells IEC-6.
Evidence-based complementary and alternative medicine : eCAM
Xu G,Ma T,Zhou C,Zhao F,Peng K,Li B
Sat Jul 16 00:00:00 UTC 2022
MG53 protein rejuvenates hUC-MSCs and facilitates their therapeutic effects in AD mice by activating Nrf2 signaling pathway.
Redox biology
Ma S,Zhou X,Wang Y,Li Z,Wang Y,Shi J,Guan F
Fri Jul 01 00:00:00 UTC 2022
High-mobility Group Box 1 Contributes to Hypoxic-Ischemic Brain Damage by Facilitating Imbalance of Microglial Polarization through RAGE-PI3K/Akt Pathway in Neonatal Rats.
International journal of medical sciences
Sun Y,Zhu X,Zhu K,Yu J,Cheng L,Hei M
Thu Dec 15 00:00:00 UTC 2022
Genistein Restricts the Epithelial Mesenchymal Transformation (EMT) and Stemness of Hepatocellular Carcinoma via Upregulating miR-1275 to Inhibit the EIF5A2/PI3K/Akt Pathway.
Biology
Yang X,Jiang W,Kong X,Zhou X,Zhu D,Kong L
Thu Sep 22 00:00:00 UTC 2022
Avasimibe Alleviates Disruption of the Airway Epithelial Barrier by Suppressing the Wnt/β-Catenin Signaling Pathway.
Frontiers in pharmacology
Zhou Z,Liang S,Zhou Z,Liu J,Meng X,Zou F,Yu C,Cai S
Tue Mar 01 00:00:00 UTC 2022
Construction, bioinformatics analysis, and validation of competitive endogenous RNA networks in ulcerative colitis.
Frontiers in genetics
Dong L,Zhang R,Huang Q,Shen Y,Li H,Yu S,Wu Q
Wed Sep 07 00:00:00 UTC 2022
Shuangyu Tiaozhi decoction alleviates non-alcoholic fatty liver disease by improving lipid deposition, insulin resistance, and inflammation in vitro and in vivo.
Frontiers in pharmacology
Yin G,Liang H,Sun W,Zhang S,Feng Y,Liang P,Chen S,Liu X,Pan W,Zhang F
Wed Dec 21 00:00:00 UTC 2022
Naringenin activates beige adipocyte browning in high fat diet-fed C57BL/6 mice by shaping the gut microbiota.
Food & function
Zhang S,Li J,Shi X,Tan X,Si Q
Mon Oct 03 00:00:00 UTC 2022
1H-NMR-based metabolomics reveals the biomarker panel and molecular mechanism of hepatocellular carcinoma progression.
Analytical and bioanalytical chemistry
Wang KX,Du GH,Qin XM,Gao L
Tue Feb 01 00:00:00 UTC 2022
The anti-MDR efficacy of YAN against A549/Taxol cells is associated with its inhibition on glycolysis and is further enhanced by 2-deoxy-d-glucose.
Chemico-biological interactions
Gao M,Yang Y,Gao Y,Liu T,Guan Q,Zhou T,Shi Y,Hao M,Li Z,Zuo D,Zhang W,Wu Y
Fri Feb 25 00:00:00 UTC 2022
Polygonum cuspidatum inhibits the growth of osteosarcoma cells via impeding Akt/ERK/EGFR signaling pathways.
Bioengineered
Zhao J,Pan B,Zhou X,Wu C,Hao F,Zhang J,Liu L
Tue Feb 01 00:00:00 UTC 2022
CaM/CaMKII mediates activation and proliferation of hepatic stellate cells regulated by ASIC1a.
Frontiers in pharmacology
Liu H,Lu WL,Hong HQ,Li MJ,Ye MP,Rao QF,Kong JL,Luan SH,Huang Y,Hu QH,Wu FR
Tue Jan 03 00:00:00 UTC 2023
Cytomegalovirus-specific neutralizing antibodies effectively prevent uncontrolled infection after allogeneic hematopoietic stem cell transplantation.
iScience
Wang H,Liu H,Zhou L,Wang D,Wang S,Liu Q,Wu Y,Tu M,Sun Z,Zheng X,Fu B,Wang B,Wei H
Fri Oct 21 00:00:00 UTC 2022
Mechanical Stimulation Protects Against Chondrocyte Pyroptosis Through Irisin-Induced Suppression of PI3K/Akt/NF-κB Signal Pathway in Osteoarthritis.
Frontiers in cell and developmental biology
Jia S,Yang Y,Bai Y,Wei Y,Zhang H,Tian Y,Liu J,Bai L
Fri Apr 01 00:00:00 UTC 2022
Saikosaponin D Inducing Apoptosis and Autophagy through the Activation of Endoplasmic Reticulum Stress in Glioblastoma.
BioMed research international
Liu G,Guan Y,Liu Y,Wang Y,Zhang J,Liu Y,Liu X
Tue Dec 06 00:00:00 UTC 2022
Artesunate Suppresses the Growth of Lung Cancer Cells by Downregulating the AKT/Survivin Signaling Pathway.
BioMed research international
Zhang W,Ning N,Huang J
Fri Apr 08 00:00:00 UTC 2022
Activation of RKIP Binding ASC Attenuates Neuronal Pyroptosis and Brain Injury via Caspase-1/GSDMD Signaling Pathway After Intracerebral Hemorrhage in Mice.
Translational stroke research
Gu L,Sun M,Li R,Tao Y,Luo X,Xu J,Wu X,Xie Z
Thu Dec 01 00:00:00 UTC 2022
N-myristoyltransferase-1 deficiency blocks myristoylation of LAMTOR1 and inhibits bladder cancer progression.
Cancer letters
Sun Y,Guan Z,Sheng Q,Duan W,Zhao H,Zhou J,Deng Q,Pei X
Thu Mar 31 00:00:00 UTC 2022
Senescent renal tubular epithelial cells activate fibroblasts by secreting Shh to promote the progression of diabetic kidney disease.
Frontiers in medicine
Wang D,Yin L,Chen R,Tan W,Liang L,Xiang J,Zhang H,Zhou X,Deng H,Guo B,Wang Y
Sat Feb 11 00:00:00 UTC 2023
miR-let-7c-3p targeting on Egr-1 contributes to the committed differentiation of leukemia cells into monocyte/macrophages.
Oncology letters
Qi F,Wang X,Zhao S,Wang C,Sun R,Wang H,Du P,Wang J,Wang X,Jiang G
Mon Aug 01 00:00:00 UTC 2022
Beta-naphthoflavone and doxorubicin synergistically enhance apoptosis in human lung cancer cells by inducing doxorubicin accumulation, mitochondrial ROS generation, and JNK pathway signaling.
Biochemical and biophysical research communications
Hoang DH,Song M,Kovale LM,Tran QH,Choe W,Kang I,Kim SS,Ha J
Tue Dec 20 00:00:00 UTC 2022
Blockage of citrate export prevents TCA cycle fragmentation via Irg1 inactivation.
Cell reports
Li Y,Li YC,Liu XT,Zhang L,Chen YH,Zhao Q,Gao W,Liu B,Yang H,Li P
Tue Feb 15 00:00:00 UTC 2022
Dioscin relieves diabetic nephropathy via suppressing oxidative stress and apoptosis, and improving mitochondrial quality and quantity control.
Food & function
Zhong Y,Liu J,Sun D,Guo T,Yao Y,Xia X,Shi C,Peng X
Mon Mar 21 00:00:00 UTC 2022
Verteporfin inhibits the dedifferentiation of tubular epithelial cells via TGF-β1/Smad pathway but induces podocyte loss in diabetic nephropathy.
Life sciences
Qi C,Hu Y,Zeng M,Chen H,Shi J,Jue H,Zhao Z,Liu J,Zhang Z,Xu Y,Wu H
Thu Dec 15 00:00:00 UTC 2022
Iron oxide nanoparticles oxidize transformed RAW 264.7 macrophages into foam cells: Impact of pulmonary surfactant component dipalmitoylphosphatidylcholine.
Chemosphere
Li H,Tao X,Song E,Song Y
Mon Aug 01 00:00:00 UTC 2022
Thrombus Inhibition and Neuroprotection for Ischemic Stroke Treatment through Platelet Regulation and ROS Scavenging.
ChemMedChem
Ye H,Ma Z,Liu L,Zhang T,Han Q,Xiang Z,Xia Y,Ke Y,Guan X,Shi Q,Ataullakhanov FI,Panteleev M
Fri Dec 16 00:00:00 UTC 2022
Colletofragarone A2 Inhibits Cancer Cell Growth In Vivo and Leads to the Degradation and Aggregation of Mutant p53.
Chemical research in toxicology
Sadahiro Y,Hitora Y,Kimura I,Hitora-Imamura N,Onodera R,Motoyama K,Tsukamoto S
Mon Sep 19 00:00:00 UTC 2022
Serine 727 phosphorylation is necessary to induce the STAT3-mediated transcription of LINC00184 in oesophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Jin C,Qi H,Xu Y,Li S
Wed Jun 01 00:00:00 UTC 2022
Titania-Nanotube-Coated Titanium Substrates Promote Osteogenesis and Suppress Osteoclastogenesis via Integrin ανβ3.
ACS applied bio materials
Kang H,Dong Y,Liu H,Luo C,Song H,Zhu M,Guo Q,Peng R,Li F,Li Y
Mon Dec 19 00:00:00 UTC 2022
Cordycepin exhibits anti-fatigue effect via activating TIGAR/SIRT1/PGC-1α signaling pathway.
Biochemical and biophysical research communications
Chai X,Pan M,Wang J,Feng M,Wang Y,Zhang Q,Sun Y
Sat Dec 31 00:00:00 UTC 2022
Sodium tanshinone IIA sulfonate enhances the BMP9-BMPR2-Smad1/5/9 signaling pathway in rat pulmonary microvascular endothelial cells and human embryonic stem cell-derived endothelial cells.
Biochemical pharmacology
Wang J,Liu W,Lu W,Luo X,Lin Y,Liu S,Qian J,Zhang C,Chen H,Li Y,Li X,Chen J,Chen Y,Jiang Q,Liu C,Hong C,Wang T,Tang H,Zhong N,Yang J,Yang K,Sun D
Sun May 01 00:00:00 UTC 2022
Molecular imaging and treatment of PSMA-positive prostate cancer with 99mTc radiolabeled aptamer-siRNA chimeras.
Nuclear medicine and biology
Jiao Y,Xu P,Luan S,Wang X,Gao Y,Zhao C,Fu P
Wed Mar 30 00:00:00 UTC 2022
High‑glucose microenvironment promotes perineural invasion of pancreatic cancer via activation of hypoxia inducible factor 1α.
Oncology reports
Zhang L,Zhang W,Zhang X,Min Y,Zhao Y,Wang B,Li W,Mao S,Min W
Fri Apr 01 00:00:00 UTC 2022
Polystyrene micro- and nano-particle coexposure injures fetal thalamus by inducing ROS-mediated cell apoptosis.
Environment international
Yang D,Zhu J,Zhou X,Pan D,Nan S,Yin R,Lei Q,Ma N,Zhu H,Chen J,Han L,Ding M,Ding Y
Mon Aug 01 00:00:00 UTC 2022
Ssc-MiR-21-5p and Ssc-MiR-615 Regulates the Proliferation and Apoptosis of Leydig Cells by Targeting SOX5.
Cells
Tang Q,Zhang Y,Yue L,Ren H,Pan C
Thu Jul 21 00:00:00 UTC 2022
Polysaccharide, the Active Component of Dendrobium officinale, Ameliorates Metabolic Hypertension in Rats via Regulating Intestinal Flora-SCFAs-Vascular Axis.
Frontiers in pharmacology
Li B,Wang HY,Huang JH,Xu WF,Feng XJ,Xiong ZP,Dong YJ,Li LZ,He X,Wu HS,Zhang K,Su J,Yu QX,Jiang NH,Lv GY,Chen SH
Fri Jul 29 00:00:00 UTC 2022
A Novel Mouse Model of Idiopathic Nephrotic Syndrome Induced by Immunization with the Podocyte Protein Crb2.
Journal of the American Society of Nephrology : JASN
Hada I,Shimizu A,Takematsu H,Nishibori Y,Kimura T,Fukutomi T,Kudo A,Ito-Nitta N,Kiuchi Z,Patrakka J,Mikami N,Leclerc S,Akimoto Y,Hirayama Y,Mori S,Takano T,Yan K
Tue Nov 01 00:00:00 UTC 2022
A Novel Mouse Model of Idiopathic Nephrotic Syndrome Induced by Immunization with the Podocyte Protein Crb2.
Journal of the American Society of Nephrology : JASN
Hada I,Shimizu A,Takematsu H,Nishibori Y,Kimura T,Fukutomi T,Kudo A,Ito-Nitta N,Kiuchi Z,Patrakka J,Mikami N,Leclerc S,Akimoto Y,Hirayama Y,Mori S,Takano T,Yan K
Tue Nov 01 00:00:00 UTC 2022
GLUT5-KHK axis-mediated fructose metabolism drives proliferation and chemotherapy resistance of colorectal cancer.
Cancer letters
Shen Z,Li Z,Liu Y,Li Y,Feng X,Zhan Y,Lin M,Fang C,Fang Y,Deng H
Sat May 28 00:00:00 UTC 2022
Holothuria Leucospilota polysaccharides alleviate hyperlipidemia via alteration of lipid metabolism and inflammation-related gene expression.
Journal of food biochemistry
Lu S,Yuan Y,Chen F,Zheng Y,Li C,Cao J,Xia G,Liu Z,Shen X,He Y,Zhou D,Zhu K
Thu Dec 01 00:00:00 UTC 2022
Acupuncture Effect Assessment in APP/PS1 Transgenic Mice: On Regulating Learning-Memory Abilities, Gut Microbiota, and Microbial Metabolites.
Computational and mathematical methods in medicine
Yang B,He M,Chen X,Sun M,Pan T,Xu X,Zhang X,Gong Q,Zhao Y,Jin Z,Cheng Z
Tue Apr 19 00:00:00 UTC 2022
Thymoquinone upregulates IL17RD in controlling the growth and metastasis of triple negative breast cancer cells in vitro.
BMC cancer
Khan MA,Zheng M,Fu J,Tania M,Li J,Fu J
Mon Jun 27 00:00:00 UTC 2022
Wnt3a knockdown promotes collagen type II expression in rat chondrocytes.
Experimental and therapeutic medicine
Shi S,Man Z,Sun S
Mon Aug 01 00:00:00 UTC 2022
Gut microbiota and butyrate contribute to nonalcoholic fatty liver disease in premenopause due to estrogen deficiency.
PloS one
Liu L,Fu Q,Li T,Shao K,Zhu X,Cong Y,Zhao X
Tue Mar 01 00:00:00 UTC 2022
Gamma-oryzanol protects human liver cell (L02) from hydrogen peroxide-induced oxidative damage through regulation of the MAPK/Nrf2 signaling pathways.
Journal of food biochemistry
Ma Y,Xiang S,Jiang W,Kong L,Tan Z,Liang Z,Yuan Z,Yi J,Zhu L
Fri Jul 01 00:00:00 UTC 2022
Ras-related protein Rab-20 inhibition alleviates cerebral ischemia/reperfusion injury by inhibiting mitochondrial fission and dysfunction.
Frontiers in molecular neuroscience
Guo J,Zhang L,Bu Y,Li W,Hu J,Li J
Fri Nov 18 00:00:00 UTC 2022
S100A8 regulates autophagy-dependent ferroptosis in microglia after experimental subarachnoid hemorrhage.
Experimental neurology
Tao Q,Qiu X,Li C,Zhou J,Gu L,Zhang L,Pang J,Zhang L,Yin S,Jiang Y,Peng J
Tue Nov 01 00:00:00 UTC 2022
Delivery of nVEGFi using AAV8 for the treatment of neovascular age-related macular degeneration.
Molecular therapy. Methods & clinical development
She K,Su J,Wang Q,Liu Y,Zhong X,Jin X,Zhao Q,Xiao J,Li R,Deng H,Lu F,Yang Y,Wei Y
Thu Mar 10 00:00:00 UTC 2022
Sestrin2 reduces cancer stemness via Wnt/β-catenin signaling in colorectal cancer.
Cancer cell international
Wei J,Zheng X,Li W,Li X,Fu Z
Fri Feb 11 00:00:00 UTC 2022
Prenatal hypothyroidism diminished exogenous NO-mediated diastolic effects in fetal rat thoracic aorta smooth muscle via increased oxidative stress.
Reproductive toxicology (Elmsford, N.Y.)
Guo J,Yu X,Liu Y,Lu L,Zhu D,Zhang Y,Li L,Zhang P,Gao Q,Lu X,Sun M
Sat Oct 01 00:00:00 UTC 2022
Adaptation to partial urethral obstruction in healthy aging LOU rats and the role of nerve growth factor signaling pathway in the bladder.
Experimental gerontology
Mossa A,Cammisotto PG,Velasquez-Flores M,Campeau L
Sat Jan 01 00:00:00 UTC 2022
A DNA damage nanoamplifier for the chemotherapy of triple-negative breast cancer via DNA damage induction and repair blocking.
International journal of pharmaceutics
Huang Y,Dai X,Guan Z,Liu D,Ren L,Chen M,Zeng Z,Jiang J,Luo Y,He Y,Huang M,Zhao C
Sat Jun 25 00:00:00 UTC 2022
Benzo(a)pyrene-7,8-dihydrodiol-9,10-epoxide induces ferroptosis in neuroblastoma cells through redox imbalance.
The Journal of toxicological sciences
Lyu Y,Ren XK,Guo CC,Li ZF,Zheng JP
Fri Dec 02 00:00:00 UTC 2022
Acute hypoxia induced dysregulation of clock-controlled ovary functions.
Frontiers in physiology
Ding M,Lu Y,Huang X,Xing C,Hou S,Wang D,Zhang Y,Wang W,Zhang C,Zhang M,Meng F,Liu K,Liu G,Zhao J,Song L
Wed Jan 11 00:00:00 UTC 2023
High expression of eIF4A1 predicts unfavorable prognosis in clear cell renal cell carcinoma.
Molecular and cellular probes
Zhang LL,Chang W,He SB,Zhang B,Ma G,Shang PF,Yue ZJ
Sat Oct 01 00:00:00 UTC 2022
Spiropachysine A suppresses hepatocellular carcinoma proliferation by inducing methuosis in vitro and in vivo.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Fang Y,Zhong T,Yang L,Luo F,Li Q,Wang D,Li Q,Fan Y,Yang X
Wed Jul 20 00:00:00 UTC 2022
FAM107A Inactivation Associated with Promoter Methylation Affects Prostate Cancer Progression through the FAK/PI3K/AKT Pathway.
Cancers
Ke S,Liu Z,Wang Q,Zhai G,Shao H,Yu X,Guo J
Sat Aug 13 00:00:00 UTC 2022
FNDC5 Promotes Adipogenic Differentiation of Primary Preadipocytes in Mashen Pigs.
Genes
Hei W,You Z,An J,Zhao T,Li J,Zhang W,Li M,Yang Y,Gao P,Cao G,Guo X,Cai C,Li B
Wed Dec 28 00:00:00 UTC 2022
Long non-coding RNA BBOX1-AS1 exacerbates esophageal squamous cell carcinoma development by regulating HOXB7/β-catenin axis.
Experimental cell research
Sheng J,Zhou M,Wang C,Jia J,Chu J,Ju C,Wan J,He J,He F
Wed Jun 01 00:00:00 UTC 2022
Matrine, a potential c-Myc inhibitor, suppresses ribosome biogenesis and nucleotide metabolism in myeloid leukemia.
Frontiers in pharmacology
Zhong WJ,Ma L,Yang F,Cao J,Tan J,Li B
Tue Nov 08 00:00:00 UTC 2022
Promotion of tumor progression induced by continuous low-dose administration of antineoplastic agent gemcitabine or gemcitabine combined with cisplatin.
Life sciences
Chen Y,Liu H,Zheng Q,Li H,You H,Feng Y,Feng W
Sat Oct 01 00:00:00 UTC 2022
Wheat oligopeptides enhance the intestinal mucosal barrier and alleviate inflammation via the TLR4/Myd88/MAPK signaling pathway in aged mice.
Food & nutrition research
Xian Y,Da P,Chao Y,Hui X,Ligang Y,Shaokang W,Guiju S
Thu Mar 10 00:00:00 UTC 2022
Microglia jointly degrade fibrillar alpha-synuclein cargo by distribution through tunneling nanotubes.
Cell
Scheiblich H,Dansokho C,Mercan D,Schmidt SV,Bousset L,Wischhof L,Eikens F,Odainic A,Spitzer J,Griep A,Schwartz S,Bano D,Latz E,Melki R,Heneka MT
Thu Sep 30 00:00:00 UTC 2021
Ataxia-telangiectasia mutated interacts with Parkin and induces mitophagy independent of kinase activity. Evidence from mantle cell lymphoma.
Haematologica
Sarkar A,Stellrecht CM,Vangapandu HV,Ayres M,Kaipparettu BA,Park JH,Balakrishnan K,Burks JK,Pandita TK,Hittelman WN,Neelapu SS,Gandhi V
Mon Feb 01 00:00:00 UTC 2021
Serum exosomal pregnancy zone protein as a promising biomarker in inflammatory bowel disease.
Cellular & molecular biology letters
Shao J,Jin Y,Shao C,Fan H,Wang X,Yang G
Tue Aug 10 00:00:00 UTC 2021
Sterol and oxysterol synthases near the ciliary base activate the Hedgehog pathway.
The Journal of cell biology
Findakly S,Daggubati V,Garcia G,LaStella SA,Choudhury A,Tran C,Li A,Tong P,Garcia JQ,Puri N,Reiter JF,Xu L,Raleigh DR
Mon Jan 04 00:00:00 UTC 2021
Human Tissue Kallikrein 1 Is Downregulated in Elderly Human Prostates and Possesses Potential In Vitro Antioxidative and Antifibrotic Effects in Rodent Prostates.
Oxidative medicine and cellular longevity
Zhang M,Luo C,Lin D,Cui K,Chen Z,Liu J
Fri Nov 12 00:00:00 UTC 2021
Mefunidone ameliorates diabetic kidney disease in STZ and db/db mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Jiang Y,Xie F,Lv X,Wang S,Liao X,Yu Y,Dai Q,Zhang Y,Meng J,Hu G,Peng Z,Tao L
Fri Jan 01 00:00:00 UTC 2021
The canonical smooth muscle cell marker TAGLN is present in endothelial cells and is involved in angiogenesis.
Journal of cell science
Tsuji-Tamura K,Morino-Koga S,Suzuki S,Ogawa M
Sun Aug 01 00:00:00 UTC 2021
Overexpression of TC2N is associated with poor prognosis in gastric cancer.
Journal of Cancer
Xu J,Ou X,Li J,Cai Q,Sun K,Ye J,Peng J
Fri Jun 17 00:00:00 UTC 2022
Aggravation of hepatic ischemia‑reperfusion injury with increased inflammatory cell infiltration is associated with the TGF‑β/Smad3 signaling pathway.
Molecular medicine reports
Li H,Shen X,Tong Y,Ji T,Feng Y,Tang Y,Mai R,Ye J,Que T,Luo X
Sun Aug 01 00:00:00 UTC 2021
Salusin‑β participates in high glucose‑induced HK‑2 cell ferroptosis in a Nrf‑2‑dependent manner.
Molecular medicine reports
Wang WJ,Jiang X,Gao CC,Chen ZW
Wed Sep 01 00:00:00 UTC 2021
Ethanol extract of Ardisiae Japonicae Herba inhibits hepatoma carcinoma cell proliferation in vitro through regulating lipid metabolism.
Chinese herbal medicines
Gong X,Cui HT,Bian YH,Li YT,Wang YX,Peng YF,Wen WB,Li K,Wang HW,Zhang ZY,Zheng F
Thu Jul 01 00:00:00 UTC 2021
Resveratrol affects the expression of uric acid transporter by improving inflammation.
Molecular medicine reports
Zhang X,Nie Q,Zhang Z,Zhao J,Zhang F,Wang C,Wang X,Song G
Sun Aug 01 00:00:00 UTC 2021
Oxygen nanocarrier broke the hypoxia trap of solid tumors and rescued transfection efficiency for gene therapy.
Journal of nanobiotechnology
Qin B,Jiang M,Li X,Shi Y,Zhang J,Luo Z,Luo L,Lu Y,Liu X,Wang S,Du Y,Qiu Y,Lou Y,You J
Sat Dec 18 00:00:00 UTC 2021
Neutralizing Antibodies and Cellular Immune Responses Against SARS-CoV-2 Sustained One and a Half Years After Natural Infection.
Frontiers in microbiology
Yan LN,Liu PP,Li XG,Zhou SJ,Li H,Wang ZY,Shen F,Lu BC,Long Y,Xiao X,Wang ZD,Li D,Han HJ,Yu H,Zhou SH,Lv WL,Yu XJ
Tue Mar 22 00:00:00 UTC 2022
Tβ4 ameliorates oxidative damage and apoptosis through ERK/MAPK and 5-HT1A signaling pathway in Aβ insulted SH-SY5Y cells.
Life sciences
Zhang GH,Pare RB,Chin KL,Qian YH
Thu Nov 25 00:00:00 UTC 2021
Cimigenoside functions as a novel γ-secretase inhibitor and inhibits the proliferation or metastasis of human breast cancer cells by γ-secretase/Notch axis.
Pharmacological research
Jia H,Liu M,Wang X,Jiang Q,Wang S,Santhanam RK,Lv C,Zhao Q,Lu J
Thu Jul 01 00:00:00 UTC 2021
Effects of photoperiod on morphology and function in testis and epididymis of Cricetulus barabensis.
Journal of cellular physiology
Mou J,Xu J,Wang Z,Wang C,Yang X,Wang X,Xue H,Wu M,Xu L
Mon Mar 01 00:00:00 UTC 2021
Glutaredoxin 1 protects neurons from oxygen-glucose deprivation/reoxygenation (OGD/R)-induced apoptosis and oxidative stress via the modulation of GSK-3β/Nrf2 signaling.
Journal of bioenergetics and biomembranes
Qiu Z,Li X,Duan C,Li R,Han L
Sun Aug 01 00:00:00 UTC 2021
Hypoxic acclimation improves cardiac redox homeostasis and protects heart against ischemia-reperfusion injury through upregulation of O-GlcNAcylation.
Redox biology
Ou W,Liang Y,Qin Y,Wu W,Xie M,Zhang Y,Zhang Y,Ji L,Yu H,Li T
Thu Jul 01 00:00:00 UTC 2021
CQMUH-011 Inhibits LPS-Induced Microglia Activation and Ameliorates Brain Ischemic Injury in Mice.
Inflammation
Liu H,Hu X,Jiang R,Cai J,Lin Q,Fan Z,Zhao P,Wang S,Zou C,Du W,Dong Z,Liu Y
Sun Aug 01 00:00:00 UTC 2021
Melatonin inhibits lipid accumulation to repress prostate cancer progression by mediating the epigenetic modification of CES1.
Clinical and translational medicine
Zhou L,Zhang C,Yang X,Liu L,Hu J,Hou Y,Tao H,Sugimura H,Chen Z,Wang L,Chen K
Tue Jun 01 00:00:00 UTC 2021
The molecular mechanism underlying mitophagy-mediated hippocampal neuron apoptosis in diabetes-related depression.
Journal of cellular and molecular medicine
Liu J,Liu L,Han YS,Yi J,Guo C,Zhao HQ,Ling J,Wang YH
Sun Aug 01 00:00:00 UTC 2021
An ESCRT-dependent step in fatty acid transfer from lipid droplets to mitochondria through VPS13D-TSG101 interactions.
Nature communications
Wang J,Fang N,Xiong J,Du Y,Cao Y,Ji WK
Tue Feb 23 00:00:00 UTC 2021
MALAT1 sponges miR-26a and miR-26b to regulate endothelial cell angiogenesis via PFKFB3-driven glycolysis in early-onset preeclampsia.
Molecular therapy. Nucleic acids
Li Q,Liu X,Liu W,Zhang Y,Wu M,Chen Z,Zhao Y,Zou L
Fri Mar 05 00:00:00 UTC 2021
miR‑4792 regulates inflammatory responses in Cryptococcus neoformans‑infected microglia.
International journal of molecular medicine
Yao G,Wang X,Wang Y,Hou Q,Gao R,Wang Y,Teng L,Lin W,Wang Z,Jin Y,Chen J
Mon Nov 01 00:00:00 UTC 2021
MicroRNA‑338‑3p regulates age‑associated osteoporosis via targeting PCSK5.
Molecular medicine reports
Tong J,Zhang M,Li X,Ren G
Mon Feb 01 00:00:00 UTC 2021
Inhibition of selenoprotein synthesis by Zika virus may contribute to congenital Zika syndrome and microcephaly by mimicking SELENOP knockout and the genetic disease PCCA.
BBA advances
Dailey GP,Premadasa LS,Ruzicka JA,Taylor EW
Thu Mar 10 00:00:00 UTC 2022
Mitochondrial calcium uniporter promotes cell proliferation and migration in esophageal cancer.
Oncology letters
Miao Y,Wang X,Lai Y,Lin W,Huang Y,Yin H,Hou R,Zhang F
Wed Sep 01 00:00:00 UTC 2021
CLIC1 Inhibition Protects Against Cellular Senescence and Endothelial Dysfunction Via the Nrf2/HO-1 Pathway.
Cell biochemistry and biophysics
Lu D,Le Y,Ding J,Dou X,Mao W,Zhu J
Tue Jun 01 00:00:00 UTC 2021
Mechanistic study of lncRNA UCA1 promoting growth and cisplatin resistance in lung adenocarcinoma.
Cancer cell international
Fu J,Pan J,Yang X,Zhang Y,Shao F,Chen J,Huang K,Wang Y
Mon Sep 20 00:00:00 UTC 2021
The SESAME complex regulates cell senescence through the generation of acetyl-CoA.
Nature metabolism
Chen W,Yu X,Wu Y,Tang J,Yu Q,Lv X,Zha Z,Hu B,Li X,Chen J,Ma L,Workman JL,Li S
Thu Jul 01 00:00:00 UTC 2021
Riclinoctaose Attenuates Renal Ischemia-Reperfusion Injury by the Regulation of Macrophage Polarization.
Frontiers in pharmacology
Zhao Y,Ding Z,Ge W,Liu J,Xu X,Cheng R,Zhang J
Tue Nov 02 00:00:00 UTC 2021
Dietary supplementation of inulin alleviates metabolism disorders in gestational diabetes mellitus mice via RENT/AKT/IRS/GLUT4 pathway.
Diabetology & metabolic syndrome
Miao M,Dai Y,Rui C,Fan Y,Wang X,Fan C,Mu J,Hou W,Dong Z,Li P,Sun G,Zeng X
Fri Dec 24 00:00:00 UTC 2021
SMAD2 regulates testicular development and testosterone synthesis in Hu sheep.
Theriogenology
Yang H,Wan Z,Jin Y,Wang F,Zhang Y
Fri Oct 15 00:00:00 UTC 2021
Endoplasmic reticulum stress manipulates autophagic response that antagonizes polybrominated diphenyl ethers quinone induced cytotoxicity in microglial BV2 cells.
Journal of hazardous materials
Zhu Q,Liu Z,Wang Y,Song E,Song Y
Sat Jun 05 00:00:00 UTC 2021
The immunosuppressive role of Edn3 overexpression in the melanoma microenvironment.
Pigment cell & melanoma research
Freitas JT,Lopez J,Llorian C,Boroni M,Kos L
Mon Nov 01 00:00:00 UTC 2021
Resolvin D1 attenuates CCl4 Induced Liver Fibrosis by Inhibiting Autophagy-Mediated HSC activation via AKT/mTOR Pathway.
Frontiers in pharmacology
Li J,Deng X,Wang S,Jiang Q,Xu K
Fri Jan 07 00:00:00 UTC 2022
Expression pattern and regulatory effect of lysine-specific demethylase 2A gene in clear cell renal cell carcinoma.
BMC urology
Wang J,Li T,Li X,Zhang Y,Wang X
Sat Aug 14 00:00:00 UTC 2021
Bone marrow macrophage-derived exosomal miR-143-5p contributes to insulin resistance in hepatocytes by repressing MKP5.
Cell proliferation
Li L,Zuo H,Huang X,Shen T,Tang W,Zhang X,An T,Dou L,Li J
Wed Dec 01 00:00:00 UTC 2021
Oxymatrine reduces expression of programmed death-ligand 1 by promoting DNA demethylation in colorectal cancer cells.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico
Hua S,Gu M,Wang Y,Ban D,Ji H
Thu Apr 01 00:00:00 UTC 2021
Let-7a regulates EV secretion and mitochondrial oxidative phosphorylation by targeting SNAP23 in colorectal cancer.
Journal of experimental & clinical cancer research : CR
Liu YD,Zhuang XP,Cai DL,Cao C,Gu QS,Liu XN,Zheng BB,Guan BJ,Yu L,Li JK,Ding HB,Yan DW
Thu Jan 14 00:00:00 UTC 2021
Ginsenoside Rg1 attenuates mechanical stress-induced cardiac injury via calcium sensing receptor-related pathway.
Journal of ginseng research
Lu ML,Wang J,Sun Y,Li C,Sun TR,Hou XW,Wang HX
Mon Nov 01 00:00:00 UTC 2021
Hsa_circ_0042823 accelerates cancer progression via miR-877-5p/FOXM1 axis in laryngeal squamous cell carcinoma.
Annals of medicine
Wu T,Sun Y,Sun Z,Li S,Wang W,Yu B,Wang G
Wed Dec 01 00:00:00 UTC 2021
Evaluation of piggyBac-mediated anti-CD19 CAR-T cells after ex vivo expansion with aAPCs or magnetic beads.
Journal of cellular and molecular medicine
Yang LR,Li L,Meng MY,Wang WJ,Yang SL,Zhao YY,Wang RQ,Gao H,Tang WW,Yang Y,Yang LL,Liao LW,Hou ZL
Fri Jan 01 00:00:00 UTC 2021
ChREBP deficiency alleviates apoptosis by inhibiting TXNIP/oxidative stress in diabetic nephropathy.
Journal of diabetes and its complications
Chen N,Song S,Yang Z,Wu M,Mu L,Zhou T,Shi Y
Wed Dec 01 00:00:00 UTC 2021
Induced Pluripotent Stem Cell-Derived Conditioned Medium Promotes Endogenous Leukemia Inhibitory Factor to Attenuate Endotoxin-Induced Acute Lung Injury.
International journal of molecular sciences
Su VY,Chiou SH,Chen WC,Yu WK,Wu HH,Chen H,Yang KY
Mon May 24 00:00:00 UTC 2021
Dual Hyaluronic Acid and Folic Acid Targeting pH-Sensitive Multifunctional 2DG@DCA@MgO-Nano-Core-Shell-Radiosensitizer for Breast Cancer Therapy.
Cancers
Askar MA,Thabet NM,El-Sayyad GS,El-Batal AI,Abd Elkodous M,El Shawi OE,Helal H,Abdel-Rafei MK
Sun Nov 07 00:00:00 UTC 2021
miR-23a/b-3p promotes hepatic lipid accumulation by regulating Srebp-1c and Fas.
Journal of molecular endocrinology
Li L,Zhang X,Ren H,Huang X,Shen T,Tang W,Dou L,Li J
Fri Nov 26 00:00:00 UTC 2021
USP15 participates in DBP-induced testicular oxidative stress injury through regulating the Keap1/Nrf2 signaling pathway.
The Science of the total environment
Zhang L,Gao X,Qin Z,Shi X,Xu K,Wang S,Tang M,Wang W,Gao S,Zuo L,Zhang L,Zhang W
Fri Aug 20 00:00:00 UTC 2021
Ubenimex Suppresses the Ability of Migration and Invasion in Gastric Cancer Cells by Alleviating the Activity of the CD13/NAB1/MAPK Pathway.
Cancer management and research
Liu X,Guo Q,Jing F,Zhou C,Xiu T,Shi Y,Jing F
Sat Apr 23 00:00:00 UTC 2022
Loss of PHLDA1 has a protective role in OGD/R-injured neurons via regulation of the GSK-3β/Nrf2 pathway.
Human & experimental toxicology
Yang F,Chen R
Mon Nov 01 00:00:00 UTC 2021
Anti-NMDAR encephalitis induced in mice by active immunization with a peptide from the amino-terminal domain of the GluN1 subunit.
Journal of neuroinflammation
Ding Y,Zhou Z,Chen J,Peng Y,Wang H,Qiu W,Xie W,Zhang J,Wang H
Sun Feb 21 00:00:00 UTC 2021
A novel chemical inhibitor suppresses breast cancer cell growth and metastasis through inhibiting HPIP oncoprotein.
Cell death discovery
Li P,Cao S,Huang Y,Zhang Y,Liu J,Cai X,Zhou L,Li J,Jiang Z,Ding L,Zheng Z,Li S,Ye Q
Thu Jul 29 00:00:00 UTC 2021
Tooth Loss Induces Memory Impairment and Gliosis in App Knock-In Mouse Models of Alzheimer's Disease.
Journal of Alzheimer's disease : JAD
Taslima F,Jung CG,Zhou C,Abdelhamid M,Abdullah M,Goto T,Saito T,Saido TC,Michikawa M
Fri Sep 17 00:00:00 UTC 2021
Self-Assembled DNA Nanostructure as a Carrier for Targeted siRNA Delivery in Glioma Cells.
International journal of nanomedicine
Zhou Y,Yang Q,Wang F,Zhou Z,Xu J,Cheng S,Cheng Y
Fri Mar 26 00:00:00 UTC 2021
Smooth muscle-specific HuR knockout induces defective autophagy and atherosclerosis.
Cell death & disease
Liu S,Jiang X,Cui X,Wang J,Liu S,Li H,Yang J,Zhang C,Zhang W
Fri Apr 09 00:00:00 UTC 2021
Treadmill exercise alleviates neuronal damage by suppressing NLRP3 inflammasome and microglial activation in the MPTP mouse model of Parkinson's disease.
Brain research bulletin
Wang W,Lv Z,Gao J,Liu M,Wang Y,Tang C,Xiang J
Wed Sep 01 00:00:00 UTC 2021
Trichostatin A increases BDNF protein expression by improving XBP-1s/ATF6/GRP78 axis in Schwann cells of diabetic peripheral neuropathy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
An J,Zhang X,Jia K,Zhang C,Zhu L,Cheng M,Li F,Zhao S,Hao J
Fri Jan 01 00:00:00 UTC 2021
LncRNA ZNFTR functions as an inhibitor in pancreatic cancer by modulating ATF3/ZNF24/VEGFA pathway.
Cell death & disease
Li W,Han S,Hu P,Chen D,Zeng Z,Hu Y,Xu F,Tang J,Wang F,Zhao Y,Huang M,Zhao G
Fri Sep 03 00:00:00 UTC 2021
Protective effect of recombinant Lactobacillus plantarum against H2O2-induced oxidative stress in HUVEC cells.
Journal of Zhejiang University. Science. B
Wang G,Hao M,Liu Q,Jiang Y,Huang H,Yang G,Wang C
Sat May 15 00:00:00 UTC 2021
Hypothermic oxygenated perfusion inhibits HECTD3-mediated TRAF3 polyubiquitination to alleviate DCD liver ischemia-reperfusion injury.
Cell death & disease
Zhou W,Zhong Z,Lin D,Liu Z,Zhang Q,Xia H,Peng S,Liu A,Lu Z,Wang Y,Ye S,Ye Q
Wed Feb 24 00:00:00 UTC 2021
Sorafenib-Induced Autophagy Promotes Glycolysis by Upregulating the p62/HDAC6/HSP90 Axis in Hepatocellular Carcinoma Cells.
Frontiers in pharmacology
Yan X,Tian R,Sun J,Zhao Y,Liu B,Su J,Li M,Sun W,Xu X
Tue Mar 08 00:00:00 UTC 2022
H2S Attenuates Sleep Deprivation-Induced Cognitive Impairment by Reducing Excessive Autophagy via Hippocampal Sirt-1 in WISTAR RATS.
Neurochemical research
Gao S,Tang YY,Jiang L,Lan F,Li X,Zhang P,Zou W,Chen YJ,Tang XQ
Sun Aug 01 00:00:00 UTC 2021
The Anti-Inflammatory Effect and Mucosal Barrier Protection of Clostridium butyricum RH2 in Ceftriaxone-Induced Intestinal Dysbacteriosis.
Frontiers in cellular and infection microbiology
Li Y,Liu M,Liu H,Sui X,Liu Y,Wei X,Liu C,Cheng Y,Ye W,Gao B,Wang X,Lu Q,Cheng H,Zhang L,Yuan J,Li M
Mon Jul 05 00:00:00 UTC 2021
Duck Plague Virus pUL48 Protein Activates the Immediate-Early Gene to Initiate the Transcription of the Virus Gene.
Frontiers in microbiology
Zhou T,Fan D,Wang M,Cheng A,Wu Y,Yang Q,Tian B,Jia R,Ou X,Mao S,Sun D,Zhang S,Zhu D,Chen S,Liu M,Zhao XX,Huang J,Gao Q,Yu Y,Zhang L
Tue Jan 11 00:00:00 UTC 2022
Disrupted eNOS activity and expression account for vasodilator dysfunction in different stage of sepsis.
Life sciences
Hu S,Pi Q,Xu X,Yan J,Guo Y,Tan W,He A,Cheng Z,Luo S,Xia Y
Fri Jan 01 00:00:00 UTC 2021
METTL3 improves cardiomyocyte proliferation upon myocardial infarction via upregulating miR-17-3p in a DGCR8-dependent manner.
Cell death discovery
Zhao K,Yang C,Zhang J,Sun W,Zhou B,Kong X,Shi J
Wed Oct 13 00:00:00 UTC 2021
Calcium Channel Protein ORAI1 Mediates TGF-β Induced Epithelial-to-Mesenchymal Transition in Colorectal Cancer Cells.
Frontiers in oncology
Kang Q,Peng X,Li X,Hu D,Wen G,Wei Z,Yuan B
Tue Jun 01 00:00:00 UTC 2021
A key role of gut microbiota-vagus nerve/spleen axis in sleep deprivation-mediated aggravation of systemic inflammation after LPS administration.
Life sciences
Zhang Y,Xie B,Chen X,Zhang J,Yuan S
Fri Jan 15 00:00:00 UTC 2021
High expression of transmembrane P24 trafficking protein 9 predicts poor prognosis in breast carcinoma.
Bioengineered
Ju G,Xu C,Zeng K,Zhou T,Zang L
Wed Dec 01 00:00:00 UTC 2021
ABIN-1 is a key regulator in RIPK1-dependent apoptosis (RDA) and necroptosis, and ABIN-1 deficiency potentiates necroptosis-based cancer therapy in colorectal cancer.
Cell death & disease
Cai J,Hu D,Sakya J,Sun T,Wang D,Wang L,Mao X,Su Z
Mon Feb 01 00:00:00 UTC 2021
RCAN1.4 attenuates renal fibrosis through inhibiting calcineurin-mediated nuclear translocation of NFAT2.
Cell death discovery
Zhang J,Chen H,Weng X,Liu H,Chen Z,Huang Q,Wang L,Liu X
Wed Oct 27 00:00:00 UTC 2021
Chorionic villus-derived mesenchymal stem cell-mediated autophagy promotes the proliferation and invasiveness of trophoblasts under hypoxia by activating the JAK2/STAT3 signalling pathway.
Cell & bioscience
Chu Y,Zhu C,Yue C,Peng W,Chen W,He G,Liu C,Lv Y,Gao G,Yao K,Han R,Hu X,Zhang Y,Ye Y
Wed Oct 13 00:00:00 UTC 2021
Upregulation of RNA cap methyltransferase RNMT drives ribosome biogenesis during T cell activation.
Nucleic acids research
Galloway A,Kaskar A,Ditsova D,Atrih A,Yoshikawa H,Gomez-Moreira C,Suska O,Warminski M,Grzela R,Lamond AI,Darzynkiewicz E,Jemielity J,Cowling VH
Fri Jul 09 00:00:00 UTC 2021
Isolation, characterization and transcriptome analysis of porcine deltacoronavirus strain HNZK-02 from Henan Province, China.
Molecular immunology
Jin XH,Zhang YF,Yuan YX,Han L,Zhang GP,Hu H
Tue Jun 01 00:00:00 UTC 2021
Giardia duodenalis extracellular vesicles regulate the proinflammatory immune response in mouse macrophages in vitro via the MAPK, AKT and NF-κB pathways.
Parasites & vectors
Zhao P,Cao L,Wang X,Li J,Dong J,Zhang N,Li X,Li S,Sun M,Zhang X,Liang M,Pu X,Gong P
Thu Jul 08 00:00:00 UTC 2021
Abnormal Glu/mGluR2/3/PI3K pathway in the hippocampal neurovascular unit leads to diabetes-related depression.
Neural regeneration research
Liu J,Han YS,Liu L,Tang L,Yang H,Meng P,Zhao HQ,Wang YH
Thu Apr 01 00:00:00 UTC 2021
The cGAS-STING signaling pathway contributes to the inflammatory response and autophagy in Aspergillus fumigatus keratitis.
Experimental eye research
Han F,Guo H,Wang L,Zhang Y,Sun L,Dai C,Wu X
Fri Jan 01 00:00:00 UTC 2021
miRNA-27a Transcription Activated by c-Fos Regulates Myocardial Ischemia-Reperfusion Injury by Targeting ATAD3a.
Oxidative medicine and cellular longevity
Bao Y,Qiao Y,Yu H,Zhang Z,Yang H,Xin X,Chen Y,Guo Y,Wu N,Jia D
Tue Jan 11 00:00:00 UTC 2022
Eradicating intracellular MRSA via targeted delivery of lysostaphin and vancomycin with mannose-modified exosomes.
Journal of controlled release : official journal of the Controlled Release Society
Yang X,Xie B,Peng H,Shi G,Sreenivas B,Guo J,Wang C,He Y
Sun Jan 10 00:00:00 UTC 2021
Identification of a Three-Glycolysis-Related lncRNA Signature Correlated With Prognosis and Metastasis in Clear Cell Renal Cell Carcinoma.
Frontiers in medicine
Li T,Tong H,Zhu J,Qin Z,Yin S,Sun Y,Liu X,He W
Fri Jan 28 00:00:00 UTC 2022
Macrophage migration inhibitory factor activates the inflammatory response in joint capsule fibroblasts following post-traumatic joint contracture.
Aging
Zhang Y,Lu S,Fan S,Xu L,Jiang X,Wang K,Cai B
Wed Feb 17 00:00:00 UTC 2021
Decreased Expression of MPC2 Contributes to Aerobic Glycolysis and Colorectal Cancer Proliferation by Activating mTOR Pathway.
Journal of immunology research
Kuerbanjiang M,Gu L,Xu C,Xu WT,Wen S,Xue H,Xu Q
Mon Nov 08 00:00:00 UTC 2021
Structural Studies of Water-Insoluble β-Glucan from Oat Bran and Its Effect on Improving Lipid Metabolism in Mice Fed High-Fat Diet.
Nutrients
Yu S,Wang J,Li Y,Wang X,Ren F,Wang X
Sat Sep 18 00:00:00 UTC 2021
Stabilizing mast cells improves acute lung injury after orthotopic liver transplantation via promotion of apoptosis in polymorphonuclear neutrophils.
American journal of physiology. Lung cellular and molecular physiology
Chen C,Zhang Z,Tan F,Meng F,Lai L,Chi X,Zhu Q
Mon Feb 01 00:00:00 UTC 2021
Inhibition of Kelch-like epichlorohydrin-related protein 1 promotes the progression and drug resistance of lung adenocarcinoma.
PeerJ
Gao H,Tang P,Ni K,Zhu L,Chen S,Zheng Y,Wan Y
Tue Apr 26 00:00:00 UTC 2022
Isovitexin Inhibits Ginkgolic Acids-Induced Inflammation Through Downregulating SHP2 Activation.
Frontiers in pharmacology
Zhang Y,Qi Z,Wang W,Wang L,Cao F,Zhao L,Fang X
Tue Aug 31 00:00:00 UTC 2021
circRPS16 Promotes Proliferation and Invasion of Hepatocellular Carcinoma by Sponging miR-876-5p to Upregulate SPINK1.
Frontiers in oncology
Lin S,Lin Y,Wu Z,Xia W,Miao C,Peng T,Zhao Z,Ji C,Mo Z,Liu X,Jian Z
Sat Oct 02 00:00:00 UTC 2021
Targeted Deletion of Loxl3 by Col2a1-Cre Leads to Progressive Hearing Loss.
Frontiers in cell and developmental biology
Liu Z,Bai X,Wan P,Mo F,Chen G,Zhang J,Gao J
Tue Jun 22 00:00:00 UTC 2021
Mechanistic insights into the anti-depressant effect of emodin: an integrated systems pharmacology study and experimental validation.
Aging
Zeng P,Wang XM,Ye CY,Su HF,Fang YY,Zhang T,Tian Q
Sat May 29 00:00:00 UTC 2021
Effects of RNA methylation N6-methyladenosine regulators on malignant progression and prognosis of melanoma.
Cancer cell international
Liu J,Zhou Z,Ma L,Li C,Lin Y,Yu T,Wei JF,Zhu L,Yao G
Thu Aug 26 00:00:00 UTC 2021
Empagliflozin protects diabetic pancreatic tissue from damage by inhibiting the activation of the NLRP3/caspase-1/GSDMD pathway in pancreatic β cells: in vitro and in vivo studies.
Bioengineered
Liu P,Zhang Z,Wang J,Zhang X,Yu X,Li Y
Wed Dec 01 00:00:00 UTC 2021
Silencing circular RNA circ_0054537 and upregulating microRNA-640 suppress malignant progression of renal cell carcinoma via regulating neuronal pentraxin-2 (NPTX2).
Bioengineered
Pei L,Lv X,Jia G,Tan X,Li M,Zhang A
Wed Dec 01 00:00:00 UTC 2021
Hypoxic tubular epithelial cells regulate the angiogenesis of HMEC-1 cells via mediation of Rab7/MMP-2 axis.
Aging
Yang Y,Wang J,Zhang Y,Hu X,Li L,Chen P
Mon Oct 25 00:00:00 UTC 2021
Prenatal High-Sucrose Diet Induced Vascular Dysfunction in Thoracic Artery of Fetal Offspring.
Molecular nutrition & food research
Feng X,Yu T,Zhang Y,Li L,Qu M,Wang J,Dong F,Zhang L,Wang F,Zhang F,Zhou X,Xu Z,Man D
Tue Jun 01 00:00:00 UTC 2021
Metformin alleviates high glucose-induced ER stress and inflammation by inhibiting the interaction between caveolin1 and AMPKα in rat astrocytes.
Biochemical and biophysical research communications
Wang G,Cui W,Chen S,Shao Z,Li Y,Wang W,Mao L,Li J,Mei X
Fri Jan 01 00:00:00 UTC 2021
Exercise reduces hyperlipidemia-induced kidney damage in apolipoprotein E-deficient mice.
Experimental and therapeutic medicine
Qian C,Yang Q,Guo L,Zhu H,You X,Liu H,Sun Y
Mon Feb 01 00:00:00 UTC 2021
Insulin injections inhibits PTZ-induced mitochondrial dysfunction, oxidative stress and neurological deficits via the SIRT1/PGC-1α/SIRT3 pathway.
Biochimica et biophysica acta. Molecular basis of disease
Cheng Y,Zeng X,Mai Q,Bai X,Jiang Y,Li J,Fan S,Ding H
Tue Jun 01 00:00:00 UTC 2021
Analysis of the microRNA and mRNA expression profile of ricin toxin-treated RAW264.7 cells reveals that miR-155-3p suppresses cell inflammation by targeting GAB2.
Toxicology letters
Liu Z,Zhang X,Dong M,Liu Z,Wang Y,Yu H,Yu K,Xu N,Liu W,Song H
Wed Sep 01 00:00:00 UTC 2021
GADD34-mediated dephosphorylation of eIF2α facilitates pseudorabies virus replication by maintaining de novo protein synthesis.
Veterinary research
Zhu T,Jiang X,Xin H,Zheng X,Xue X,Chen JL,Qi B
Mon Dec 20 00:00:00 UTC 2021
BOP1 Silencing Suppresses Gastric Cancer Proliferation through p53 Modulation.
Current medical science
Yang YP,Qin RH,Zhao JJ,Qin XY
Thu Apr 01 00:00:00 UTC 2021
Subtractive CRISPR screen identifies the ATG16L1/vacuolar ATPase axis as required for non-canonical LC3 lipidation.
Cell reports
Ulferts R,Marcassa E,Timimi L,Lee LC,Daley A,Montaner B,Turner SD,Florey O,Baillie JK,Beale R
Tue Oct 26 00:00:00 UTC 2021
Nicotinamide improves NAD+ levels to protect against acetaminophen-induced acute liver injury in mice.
Human & experimental toxicology
Xu J,Zhang L,Jiang R,Hu K,Hu D,Liao C,Jiang S,Yang Y,Huang J,Tang L,Li L
Mon Nov 01 00:00:00 UTC 2021
Dihydroxyeicosatrienoic Acid, a Metabolite of Epoxyeicosatrienoic Acids Upregulates Endothelial Nitric Oxide Synthase Expression Through Transcription: Mechanism of Vascular Endothelial Function Protection.
Cell biochemistry and biophysics
Zuo D,Pi Q,Shi Y,Luo S,Xia Y
Tue Jun 01 00:00:00 UTC 2021
Huaier Inhibits Proliferation, Migration, and Invasion of Cutaneous Squamous Cell Carcinoma Cells by Inhibiting the Methylation Levels of CDKN2A and TP53.
Integrative cancer therapies
Wang L,Xu L,Wang Y
Mon Oct 11 00:00:00 UTC 2021
Proteomics identifies a novel role of fibrinogen-like protein 1 in Crohn's disease.
World journal of gastroenterology
Sun XL,Qiao LC,Gong J,Wen K,Xu ZZ,Yang BL
Tue Sep 21 00:00:00 UTC 2021
Curcumin suppresses colorectal tumorigenesis via the Wnt/β-catenin signaling pathway by downregulating Axin2.
Oncology letters
Hao J,Dai X,Gao J,Li Y,Hou Z,Chang Z,Wang Y
Mon Mar 01 00:00:00 UTC 2021
MiR-130a-3p suppresses colorectal cancer growth by targeting Wnt Family Member 1 (WNT1).
Bioengineered
Song GL,Xiao M,Wan XY,Deng J,Ling JD,Tian YG,Li M,Yin J,Zheng RY,Tang Y,Liu GY
Wed Dec 01 00:00:00 UTC 2021
Compound Kushen Injection intervenes metabolic reprogramming and epithelial-mesenchymal transition of HCC via regulating β-catenin/c-Myc signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang KX,Du GH,Qin XM,Gao L
Wed Dec 01 00:00:00 UTC 2021
Effect of Sirolimus on the Level of Peripheral Blood Lymphocyte Autophagy in Children With Systemic Lupus Erythematosus.
Frontiers in pediatrics
Wang X,Su Q,Dou Z,Zhao X,Zhang N,Yu B,Wang Y,Rong Z
Tue Nov 02 00:00:00 UTC 2021
Lycopene-Loaded Microemulsion Regulates Neurogenesis in Rats with Aβ-Induced Alzheimer's Disease Rats Based on the Wnt/β-catenin Pathway.
Neural plasticity
Ning WJ,Lv RJ,Xu N,Hou XY,Shen C,Guo YL,Fan ZY,Cao N,Liu XP
Fri Feb 04 00:00:00 UTC 2022
Melatonin alleviated oxidative stress induced by energy restriction on sheep Leydig cells through Sirt1/Sod2 pathway.
Theriogenology
Ma J,Yang H,Liu L,Wan Y,Wang F
Fri Oct 01 00:00:00 UTC 2021
PTIP Inhibits Cell Invasion in Esophageal Squamous Cell Carcinoma via Modulation of EphA2 Expression.
Frontiers in oncology
Han X,Zhu Y,Shen L,Zhou Y,Pang L,Zhou W,Gu H,Han K,Yang Y,Jiang C,Xie J,Zhang C,Ding L
Sat Apr 10 00:00:00 UTC 2021
SNAPIN Regulates Cell Cycle Progression to Promote Pancreatic β Cell Growth.
Frontiers in endocrinology
Jiang M,Kuang Z,He Y,Cao Y,Yu T,Cheng J,Liu W,Wang W
Thu Dec 16 00:00:00 UTC 2021
SNAPIN Regulates Cell Cycle Progression to Promote Pancreatic β Cell Growth.
Frontiers in endocrinology
Jiang M,Kuang Z,He Y,Cao Y,Yu T,Cheng J,Liu W,Wang W
Thu Dec 16 00:00:00 UTC 2021
SNAPIN Regulates Cell Cycle Progression to Promote Pancreatic β Cell Growth.
Frontiers in endocrinology
Jiang M,Kuang Z,He Y,Cao Y,Yu T,Cheng J,Liu W,Wang W
Thu Dec 16 00:00:00 UTC 2021
Resveratrol attenuates manganese-induced oxidative stress and neuroinflammation through SIRT1 signaling in mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Cong L,Lei MY,Liu ZQ,Liu ZF,Ma Z,Liu K,Li J,Deng Y,Liu W,Xu B
Thu Jul 01 00:00:00 UTC 2021
JaponiconeA induces apoptosis of bortezomib-sensitive and -resistant myeloma cells in vitro and in vivo by targeting IKK.
Cancer biology & medicine
Zhang Z,Ye C,Liu J,Xu W,Wu C,Yu Q,Xu X,Zeng X,Jin H,Wu Y,Yan H
Tue Sep 28 00:00:00 UTC 2021
KDM6A promotes imatinib resistance through YY1-mediated transcriptional upregulation of TRKA independently of its demethylase activity in chronic myelogenous leukemia.
Theranostics
Zhang C,Shen L,Zhu Y,Xu R,Deng Z,Liu X,Ding Y,Wang C,Shi Y,Bei L,Wei D,Thorne RF,Zhang XD,Yu L,Chen S
Tue Jul 27 00:00:00 UTC 2021
Rufinamide (RUF) suppresses inflammation and maintains the integrity of the blood-brain barrier during kainic acid-induced brain damage.
Open life sciences
Yu H,He B,Han X,Yan T
Tue Sep 14 00:00:00 UTC 2021
Doxorubicin and PD-L1 siRNA co-delivery with stem cell membrane-coated polydopamine nanoparticles for the targeted chemoimmunotherapy of PCa bone metastases.
Nanoscale
Mu X,Zhang M,Wei A,Yin F,Wang Y,Hu K,Jiang J
Fri May 21 00:00:00 UTC 2021
3'HS1 CTCF binding site in human β-globin locus regulates fetal hemoglobin expression.
eLife
Himadewi P,Wang XQD,Feng F,Gore H,Liu Y,Yu L,Kurita R,Nakamura Y,Pfeifer GP,Liu J,Zhang X
Wed Sep 29 00:00:00 UTC 2021
Circular RNA circEsyt2 regulates vascular smooth muscle cell remodeling via splicing regulation.
The Journal of clinical investigation
Gong X,Tian M,Cao N,Yang P,Xu Z,Zheng S,Liao Q,Chen C,Zeng C,Jose PA,Wang DZ,Jian Z,Xiao Y,Jiang DS,Wei X,Zhang B,Wang Y,Chen K,Wu G,Zeng C
Wed Dec 15 00:00:00 UTC 2021
TM9SF4 is a novel regulator in lineage commitment of bone marrow mesenchymal stem cells to either osteoblasts or adipocytes.
Stem cell research & therapy
Yu L,Xie M,Zhang F,Wan C,Yao X
Sat Nov 13 00:00:00 UTC 2021
TRIM21-regulated Annexin A2 plasma membrane trafficking facilitates osteosarcoma cell differentiation through the TFEB-mediated autophagy.
Cell death & disease
Zhang HT,Zeng Q,Wu B,Lu J,Tong KL,Lin J,Liu QY,Xu L,Yang J,Liu X,Liu W,Zhang YF,Lian Q,Liu L,Gao X
Wed Jan 06 00:00:00 UTC 2021
Chronic Intermittent Hypobaric Hypoxia Decreases High Blood Pressure by Stabilizing the Vascular Renin-Angiotensin System in Spontaneously Hypertensive Rats.
Frontiers in physiology
Chen H,Yu B,Guo X,Hua H,Cui F,Guan Y,Tian Y,Zhang X,Zhang Y,Ma H
Fri Apr 22 00:00:00 UTC 2022
Nintedanib Regulates GRK2 and CXCR2 to Reduce Neutrophil Recruitment in Endotoxin-Induced Lung Injury.
International journal of molecular sciences
Su VY,Chen WC,Yu WK,Wu HH,Chen H,Yang KY
Mon Sep 13 00:00:00 UTC 2021
CircMETTL3, upregulated in a m6A-dependent manner, promotes breast cancer progression.
International journal of biological sciences
Li Z,Yang HY,Dai XY,Zhang X,Huang YZ,Shi L,Wei JF,Ding Q
Thu Mar 24 00:00:00 UTC 2022
Bushen Huoxue recipe attenuates early pregnancy loss via activating endometrial COX2-PGE2 angiogenic signaling in mice.
BMC complementary medicine and therapies
Song Y,Zhou F,Tan X,Liu X,Ding J,Zhang C,Li F,Zhu W,Ma W,Hu R,Zhang M
Thu Jan 14 00:00:00 UTC 2021
The novel prognostic risk factor STC2 can regulate the occurrence and progression of osteosarcoma via the glycolytic pathway.
Biochemical and biophysical research communications
Yu B,Zhang F,Liu L,Liang Y,Tang X,Peng Y,Cai F,Zeng D,Yuan X,Li J,Guo Y,Lv B,Wang M,Liao Q,Lv XB
Fri May 21 00:00:00 UTC 2021
M2 Macrophage-derived exosomal miR-501 contributes to pubococcygeal muscle regeneration.
International immunopharmacology
Zhou M,Li B,Liu C,Hu M,Tang J,Min J,Cheng J,Hong L
Wed Dec 01 00:00:00 UTC 2021
cAMP-PKA signaling is involved in regulation of spinal HCN channels function in diabetic neuropathic pain.
Neuroscience letters
Ma Y,Chen J,Yu D,Wei B,Jin H,Zeng J,Liu X
Sat Apr 17 00:00:00 UTC 2021
CIHH protects the heart against left ventricular remodelling and myocardial fibrosis by balancing the renin-angiotensin system in SHR.
Life sciences
Yu B,Chen H,Guo XQ,Hua H,Guan Y,Cui F,Tian YM,Zhang HX,Zhang XJ,Zhang Y,Ma HJ
Sun Aug 01 00:00:00 UTC 2021
LINC00511 drives invasive behavior in hepatocellular carcinoma by regulating exosome secretion and invadopodia formation.
Journal of experimental & clinical cancer research : CR
Peng X,Li X,Yang S,Huang M,Wei S,Ma Y,Li Y,Wu B,Jin H,Li B,Tang S,Fan Q,Liu J,Yang L,Li H
Fri Jun 04 00:00:00 UTC 2021
EphA2 inhibits SRA01/04 cells apoptosis by suppressing autophagy via activating PI3K/Akt/mTOR pathway.
Archives of biochemistry and biophysics
Han J,Wang L,Lv H,Liu J,Dong Y,Shi L,Ji Q
Sat Oct 30 00:00:00 UTC 2021
Downregulation of circular RNA circDOCK7 identified from diabetic rats after sleeve gastrectomy contributes to hepatocyte apoptosis through regulating miR-139-3p and MCM3.
Biochemical and biophysical research communications
Li L,Dong S,Li S,Xu Q,Wang S,Xiong Y,Cheng Y,Zhong M,Zhang G,Hu S
Fri Apr 09 00:00:00 UTC 2021
Identification of candidate biomarker EMP3 and its prognostic potential in clear cell renal cell carcinoma.
Frontiers in bioscience (Landmark edition)
Lv Q,Xiao W,Xiong Z,Shi J,Miao D,Meng X,Yuan H,Yang H,Zhang X
Tue Nov 30 00:00:00 UTC 2021
Expression, purification and characterisation of a human anti-CDK4 single-chain variable fragment antibody.
BMC biotechnology
Zhao J,Xu J,Yang T,Yu X,Cheng C,Zhang T,Ren Z,Li N,Yang F,Li G
Mon Dec 20 00:00:00 UTC 2021
Geniposide from Gardenia jasminoides var. radicans Makino Attenuates Myocardial Injury in Spontaneously Hypertensive Rats via Regulating Apoptotic and Energy Metabolism Signalling Pathway.
Drug design, development and therapy
Hou Y,Yuan P,Fu Y,Zhang Q,Gao L,Wei Y,Zheng X,Feng W
Thu Oct 07 00:00:00 UTC 2021
Exploring the Role and Mechanism of pAMPKα-Mediated Dysregulation of Brf1 and RNA Pol III Genes.
Oxidative medicine and cellular longevity
Wu T,Zhang D,Lin M,Yu L,Dai T,Li S,Yu F,Lu L,Zheng L,Zhong S
Fri Nov 12 00:00:00 UTC 2021
UHRF2 commissions the completion of DNA demethylation through allosteric activation by 5hmC and K33-linked ubiquitination of XRCC1.
Molecular cell
Liu X,Xu B,Yang J,He L,Zhang Z,Cheng X,Yu H,Liu X,Jin T,Peng Y,Huang Y,Xia L,Wang Y,Wu J,Wu X,Liu S,Shan L,Yang X,Sun L,Liang J,Zhang Y,Shang Y
Thu Jul 15 00:00:00 UTC 2021
Neuroinflammation induces anxiety- and depressive-like behavior by modulating neuronal plasticity in the basolateral amygdala.
Brain, behavior, and immunity
Zheng ZH,Tu JL,Li XH,Hua Q,Liu WZ,Liu Y,Pan BX,Hu P,Zhang WH
Fri Jan 01 00:00:00 UTC 2021
LINC00839 Regulates Proliferation, Migration, Invasion, Apoptosis and Glycolysis in Neuroblastoma Cells Through miR-338-3p/GLUT1 Axis.
Neuropsychiatric disease and treatment
Yang L,Pei L,Yi J
Sun Apr 24 00:00:00 UTC 2022
CTRP1 Attenuates Cerebral Ischemia/Reperfusion Injury via the PERK Signaling Pathway.
Frontiers in cell and developmental biology
Fei H,Xiang P,Luo W,Tan X,Gu C,Liu M,Chen M,Wang Q,Yang J
Tue Aug 24 00:00:00 UTC 2021
Depiction of Aging-Based Molecular Phenotypes With Diverse Clinical Prognosis and Immunological Features in Gastric Cancer.
Frontiers in medicine
He F,Ding H,Zhou Y,Wang Y,Xie J,Yang S,Zhu Y
Sun May 01 00:00:00 UTC 2022
DNMT1-mediated PPARα methylation aggravates damage of retinal tissues in diabetic retinopathy mice.
Biological research
Zhu Y,Wang X,Zhou X,Ding L,Liu D,Xu H
Fri Aug 06 00:00:00 UTC 2021
Sinapic Acid Reduces Oxidative Stress and Pyroptosis via Inhibition of BRD4 in Alcoholic Liver Disease.
Frontiers in pharmacology
Chu J,Yan R,Wang S,Li G,Kang X,Hu Y,Lin M,Shan W,Zhao Y,Wang Z,Sun R,Yao J,Zhang N
Tue Jun 22 00:00:00 UTC 2021
Protein arginine methyltransferase 5 mediates THP-1-derived macrophage activation dependent on NF-κB in endometriosis.
Experimental and therapeutic medicine
Chai X,Wu X,He L,Ding H
Wed Sep 01 00:00:00 UTC 2021
NPM1 mutant maintains ULK1 protein stability via TRAF6-dependent ubiquitination to promote autophagic cell survival in leukemia.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Tang Y,Tao Y,Wang L,Yang L,Jing Y,Jiang X,Lei L,Yang Z,Wang X,Peng M,Xiao Q,Ren J,Zhang L
Mon Feb 01 00:00:00 UTC 2021
Azathioprine pretreatment ameliorates myocardial ischaemia reperfusion injury in diabetic rats by reducing oxidative stress, apoptosis, and inflammation.
Clinical and experimental pharmacology & physiology
Lu C,Liu L,Chen S,Niu J,Li S,Xie W,Cheng X
Wed Dec 01 00:00:00 UTC 2021
Exercise Intervention Mitigates Pathological Liver Changes in NAFLD Zebrafish by Activating SIRT1/AMPK/NRF2 Signaling.
International journal of molecular sciences
Zou Y,Chen Z,Sun C,Yang D,Zhou Z,Peng X,Zheng L,Tang C
Sun Oct 10 00:00:00 UTC 2021
Secretagogin Mediates the Regulatory Effect of Electroacupuncture on Hypothalamic-Pituitary-Adrenal Axis Dysfunction in Surgical Trauma.
Neural plasticity
Zhang M,Sun J,Wang Y,Tian Z
Mon Nov 01 00:00:00 UTC 2021
Sunitinib malate inhibits intestinal tumor development in male ApcMin/+ mice by down-regulating inflammation-related factors with suppressing β-cateinin/c-Myc pathway and re-balancing Bcl-6 and Caspase-3.
International immunopharmacology
Chen L,Xu P,Xiao Q,Chen L,Li S,Jian JM,Zhong YB
Fri Jan 01 00:00:00 UTC 2021
Arsenic Trioxide Combining Leflunomide Activates Nrf2-ARE-HO-1 Signaling Pathway and Protects Heart Xenografts.
Chinese journal of integrative medicine
Wang TD,Xu SL,Yu ZY,Ni SB,Zhang C,Jiao ZX
Fri Oct 01 00:00:00 UTC 2021
CDCA4 suppresses epithelial-mesenchymal transtion (EMT) and metastasis in Non-small cell lung cancer through modulating autophagy.
Cancer cell international
Xu C,Cao H,Sui Y,Zhang H,Shi C,Wu J,Ma R,Feng J
Tue Jan 12 00:00:00 UTC 2021
Anoctamin 5 promotes osteosarcoma development by increasing degradation of Nel-like proteins 1 and 2.
Aging
Pan R,Lu Q,Ren C,Li H,Zeng F,Tian X,Chen H
Fri Jul 09 00:00:00 UTC 2021
Yi-Qi-Jian-Pi Formula Suppresses RIPK1/RIPK3-Complex-Dependent Necroptosis of Hepatocytes Through ROS Signaling and Attenuates Liver Injury in Vivo and in Vitro.
Frontiers in pharmacology
Wang F,Tang L,Liang B,Jin C,Gao L,Li Y,Li Z,Shao J,Zhang Z,Tan S,Zhang F,Zheng S
Thu May 13 00:00:00 UTC 2021
Autophagy impairment as a key feature for acetaminophen-induced ototoxicity.
Cell death & disease
Zhao T,Zheng T,Yu H,Hu BH,Hu B,Ma P,Yang Y,Yang N,Hu J,Cao T,Chen G,Yan B,Peshoff M,Hatzoglou M,Geng R,Li B,Zheng QY
Mon Jan 04 00:00:00 UTC 2021
Internal Disulfide Bonding and Glycosylation of Interleukin-7 Protect Against Proteolytic Inactivation by Neutrophil Metalloproteinases and Serine Proteases.
Frontiers in immunology
Vandooren J,Pereira RVS,Ugarte-Berzal E,Rybakin V,Noppen S,Stas MR,Bernaerts E,Ganseman E,Metzemaekers M,Schols D,Proost P,Opdenakker G
Tue Dec 21 00:00:00 UTC 2021
Exposure to Melamine cyanuric acid in adolescent mice caused emotional disorder and behavioral disorder.
Ecotoxicology and environmental safety
Wang S,Zhang J,Zhang S,Shi F,Feng D,Feng X
Mon Mar 15 00:00:00 UTC 2021
Suppressing Sart1 to modulate macrophage polarization by siRNA-loaded liposomes: a promising therapeutic strategy for pulmonary fibrosis.
Theranostics
Pan T,Zhou Q,Miao K,Zhang L,Wu G,Yu J,Xu Y,Xiong W,Li Y,Wang Y
Thu Jul 22 00:00:00 UTC 2021
Aloe-emodin prevents nerve injury and neuroinflammation caused by ischemic stroke via the PI3K/AKT/mTOR and NF-κB pathway.
Food & function
Xian M,Cai J,Zheng K,Liu Q,Liu Y,Lin H,Liang S,Wang S
Tue Sep 07 00:00:00 UTC 2021
Curcumin Inhibits Replication of Human Parainfluenza Virus Type 3 by Affecting Viral Inclusion Body Formation.
BioMed research international
Zhang C,Zhang K,Zang G,Chen T,Lu N,Wang S,Zhang G
Tue Sep 28 00:00:00 UTC 2021
Nrf2 overexpression increases risk of high tumor mutation burden in acute myeloid leukemia by inhibiting MSH2.
Cell death & disease
Liu P,Ma D,Wang P,Pan C,Fang Q,Wang J
Tue Jan 05 00:00:00 UTC 2021
Aureonitol Analogues and Orsellinic Acid Esters Isolated from Chaetomium elatum and Their Antineuroinflammatory Activity.
Journal of natural products
Ju F,Kuang QX,Li QZ,Huang LJ,Guo WX,Gong LQ,Dai YF,Wang L,Gu YC,Wang D,Deng Y,Guo DL
Fri Dec 24 00:00:00 UTC 2021
Effects of cannabinoid (CBD) on blood brain barrier permeability after brain injury in rats.
Brain research
Jiang H,Li H,Cao Y,Zhang R,Zhou L,Zhou Y,Zeng X,Wu J,Wu D,Wu D,Guo X,Li X,Wu H,Li P
Fri Oct 01 00:00:00 UTC 2021
The Protective Effects of Sulforaphane on High-Fat Diet-Induced Obesity in Mice Through Browning of White Fat.
Frontiers in pharmacology
Liu Y,Fu X,Chen Z,Luo T,Zhu C,Ji Y,Bian Z
Tue May 18 00:00:00 UTC 2021
Gut microbiota-dependent catabolites of tryptophan play a predominant role in the protective effects of turmeric polysaccharides against DSS-induced ulcerative colitis.
Food & function
Yang C,Du Y,Ren D,Yang X,Zhao Y
Tue Oct 19 00:00:00 UTC 2021
UM-6 induces autophagy and apoptosis via the Hippo-YAP signaling pathway in cervical cancer.
Cancer letters
Wang D,He J,Dong J,Wu S,Liu S,Zhu H,Xu T
Thu Oct 28 00:00:00 UTC 2021
Aroylhydrazone Diorganotin Complexes Causes DNA Damage and Apoptotic Cell Death: From Chemical Synthesis to Biochemical Effects.
International journal of molecular sciences
Jiang W,Tan Y,Peng Y
Thu Dec 16 00:00:00 UTC 2021
Berberine Reduces Renal Cell Pyroptosis in Golden Hamsters with Diabetic Nephropathy through the Nrf2-NLRP3-Caspase-1-GSDMD Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ding B,Geng S,Hou X,Ma X,Xu H,Yang F,Liu K,Liang W,Ma G
Thu Aug 18 00:00:00 UTC 2022
Advanced maternal age causes premature placental senescence and malformation via dysregulated α-Klotho expression in trophoblasts.
Aging cell
Chen Z,Xiong L,Jin H,Yu J,Li X,Fu H,Wen L,Qi H,Tong C,Saffery R,Kilby MD,Baker PN
Thu Jul 01 00:00:00 UTC 2021
SEPHS1 promotes SMAD2/3/4 expression and hepatocellular carcinoma cells invasion.
Experimental hematology & oncology
Yang S,Zhang H,Yang H,Zhang J,Wang J,Luo T,Jiang Y,Hua H
Tue Feb 23 00:00:00 UTC 2021
YWHAZ interacts with DAAM1 to promote cell migration in breast cancer.
Cell death discovery
Mei J,Liu Y,Yu X,Hao L,Ma T,Zhan Q,Zhang Y,Zhu Y
Fri Aug 27 00:00:00 UTC 2021
FLI1 Induces Megakaryopoiesis Gene Expression Through WAS/WIP-Dependent and Independent Mechanisms; Implications for Wiskott-Aldrich Syndrome.
Frontiers in immunology
Wang C,Sample KM,Gajendran B,Kapranov P,Liu W,Hu A,Zacksenhaus E,Li Y,Hao X,Ben-David Y
Mon Jul 05 00:00:00 UTC 2021
Verapamil ameliorates proximal tubular epithelial cells apoptosis and fibrosis in diabetic kidney.
European journal of pharmacology
Song Y,Guo F,Zhao Y,Zhao L,Fan X,Zhang Y,Liu Y,Qin G
Mon Nov 15 00:00:00 UTC 2021
Phenotypic differences in the inner ears of CBA/CaJ and C57BL/6J mice carrying missense and single base pair deletion mutations in the Cdh23 gene.
Journal of neuroscience research
Zhao T,Ma P,Zhao F,Zheng T,Yan B,Zhang Q,Yuan J,Hu B,Yang Y,Hu J,Geng R,Hu BH,Sun T,Zheng QY,Li B
Fri Oct 01 00:00:00 UTC 2021
Anti-renal interstitial fibrosis effect of norcantharidin is exerted through inhibition of PP2Ac-mediated C-terminal phosphorylation of Smad3.
Chemical biology & drug design
Luo HW,Yin DD,Xiao Z,Wen L,Liao YJ,Tang CY,Zeng D,Xiao HT,Li Y
Mon Feb 01 00:00:00 UTC 2021
Colonic Epithelial PHLPP2 Deficiency Promotes Colonic Epithelial Pyroptosis by Activating the NF-κB Signaling Pathway.
Oxidative medicine and cellular longevity
Li DF,Chang X,Zhao JL,Chen XM,Xu ZL,Zhang DG,Wu BH,Wang LS,Bai Y,Yao J
Wed Jan 05 00:00:00 UTC 2022
Nogo-B receptor is required for stabilizing TGF-β type I receptor and promotes the TGF-β1-induced epithelial-to-mesenchymal transition of non-small cell lung cancer.
Journal of Cancer
Wu D,Zhao B,Song Y,Chi X,Fu H,Guan T,Zhang L,Yang X,Hu K,Huang R,Jin X,Miao QR,Shao S
Mon Jan 11 00:00:00 UTC 2021
Overexpression of Annexin A2 promotes proliferation by forming a Glypican 1/c-Myc positive feedback loop: prognostic significance in human glioma.
Cell death & disease
Li X,Nie S,Lv Z,Ma L,Song Y,Hu Z,Hu X,Liu Z,Zhou G,Dai Z,Song T,Liu J,Wang S
Fri Mar 12 00:00:00 UTC 2021
Sodium alginate/collagen hydrogel loaded with human umbilical cord mesenchymal stem cells promotes wound healing and skin remodeling.
Cell and tissue research
Zhang Z,Li Z,Li Y,Wang Y,Yao M,Zhang K,Chen Z,Yue H,Shi J,Guan F,Ma S
Mon Feb 01 00:00:00 UTC 2021
Abnormal spatiotemporal expression pattern of progranulin and neurodevelopment impairment in VPA-induced ASD rat model.
Neuropharmacology
Lan J,Hu Y,Wang X,Zheng W,Liao A,Wang S,Li Y,Wang Y,Yang F,Chen D
Wed Sep 15 00:00:00 UTC 2021
SPP1 Promotes Enzalutamide Resistance and Epithelial-Mesenchymal-Transition Activation in Castration-Resistant Prostate Cancer via PI3K/AKT and ERK1/2 Pathways.
Oxidative medicine and cellular longevity
Pang X,Zhang J,He X,Gu Y,Qian BZ,Xie R,Yu W,Zhang X,Li T,Shi X,Zhou Y,Cui Y
Mon Feb 07 00:00:00 UTC 2022
Tryptophanyl tRNA Synthetase from Human Macrophages Infected by Porphyromonas gingivalis Induces a Proinflammatory Response Associated with Atherosclerosis.
Pathogens (Basel, Switzerland)
Sasaki M,Shimoyama Y,Kodama Y,Ishikawa T
Mon Dec 20 00:00:00 UTC 2021
Calcium alleviates fluoride-induced kidney damage via FAS/FASL, TNFR/TNF, DR5/TRAIL pathways in rats.
Ecotoxicology and environmental safety
Li H,Fan J,Zhao Y,Yang J,Xu H,Manthari RK,Cheng X,Wang J,Wang J
Wed Dec 15 00:00:00 UTC 2021
A methylation-driven gene panel predicts survival in patients with colon cancer.
FEBS open bio
Peng Y,Zhao J,Yin F,Sharen G,Wu Q,Chen Q,Sun X,Yang J,Wang H,Zhang D
Wed Sep 01 00:00:00 UTC 2021
Fructooligosaccharide supplementation alleviated the pathological immune response and prevented the impairment of intestinal barrier in DSS-induced acute colitis mice.
Food & function
Liao M,Zhang Y,Qiu Y,Wu Z,Zhong Z,Zeng X,Zeng Y,Xiong L,Wen Y,Liu R
Tue Oct 19 00:00:00 UTC 2021
Transcriptomic Profiling Reveals the Antiapoptosis and Antioxidant Stress Effects of Fos in Ischemic Stroke.
Frontiers in neurology
Mu Q,Zhang Y,Gu L,Gerner ST,Qiu X,Tao Q,Pang J,Dipritu G,Zhang L,Yin S,Jiang Y,Peng J
Wed Nov 10 00:00:00 UTC 2021
A metabolic data-driven systems pharmacology strategy for decoding and validating the mechanism of Compound Kushen Injection against HCC.
Journal of ethnopharmacology
Wang KX,Chen YP,Lu AP,Du GH,Qin XM,Guan DG,Gao L
Mon Jun 28 00:00:00 UTC 2021
Metformin Corrects Glucose Metabolism Reprogramming and NLRP3 Inflammasome-Induced Pyroptosis via Inhibiting the TLR4/NF-κB/PFKFB3 Signaling in Trophoblasts: Implication for a Potential Therapy of Preeclampsia.
Oxidative medicine and cellular longevity
Zhang Y,Liu W,Zhong Y,Li Q,Wu M,Yang L,Liu X,Zou L
Tue Feb 08 00:00:00 UTC 2022
DCI after Aneurysmal Subarachnoid Hemorrhage Is Related to the Expression of MFG-E8.
BioMed research international
Chen X,Jiang Y,Liu J,Wang C,Wan D,Liang A,Leng J,Yang Y,Xiang H,Liu R
Fri Jan 14 00:00:00 UTC 2022
Interleukin-1 induces receptor activator of nuclear factor-κB ligand-independent osteoclast differentiation in RAW264.7 cells.
Experimental and therapeutic medicine
Liao R,Feng Z,Li W,Liu R,Xu X,Yao S,Tian J
Tue Jun 01 00:00:00 UTC 2021
The Role of Nrf2 on the Cognitive Dysfunction of High-fat Diet Mice Following Lead Exposure.
Biological trace element research
Zhang L,Bo J,Chen W,Li S,Wang Y,Yan L,Wu L,Zhang Y
Tue Jun 01 00:00:00 UTC 2021
Fyn kinase regulates dopaminergic neuronal apoptosis in animal and cell models of high glucose (HG) treatment.
BMC molecular and cell biology
Tan C,Liu X,Zhang X,Peng W,Wang H,Zhou W,Jiang J,Mo L,Chen Y,Chen L
Sat Dec 04 00:00:00 UTC 2021
Prenylated xanthones from mangosteen (Garcinia mangostana) activate the AhR and Nrf2 pathways and protect intestinal barrier integrity in HT-29 cells.
Free radical biology & medicine
Tocmo R,Le B,Heun A,van Pijkeren JP,Parkin K,Johnson JJ
Mon Feb 01 00:00:00 UTC 2021
Extracellular DJ-1 induces sterile inflammation in the ischemic brain.
PLoS biology
Nakamura K,Sakai S,Tsuyama J,Nakamura A,Otani K,Kurabayashi K,Yogiashi Y,Masai H,Shichita T
Sat May 01 00:00:00 UTC 2021
The role of alcohol dehydrogenase 1C in regulating inflammatory responses in ulcerative colitis.
Biochemical pharmacology
Song F,Zhang Y,Pan Z,Hu X,Zhang Q,Huang F,Ye X,Huang P
Fri Oct 01 00:00:00 UTC 2021
Sodium Tanshinone IIA Sulfonate Attenuates Cigarette Smoke Extract-Induced Mitochondrial Dysfunction, Oxidative Stress, and Apoptosis in Alveolar Epithelial Cells by Enhancing SIRT1 Pathway.
Toxicological sciences : an official journal of the Society of Toxicology
Guan R,Yao H,Li Z,Qian J,Yuan L,Cai Z,Ding M,Liu W,Xu J,Li Y,Sun D,Wang J,Lu W
Tue Sep 28 00:00:00 UTC 2021
Downregulating lncRNA PRNCR1 ameliorates LPS-induced pulmonary vascular endothelial cell injury by modulating miR-330-5p/TLR4 axis.
Journal of biochemical and molecular toxicology
Yu Y,Sun H,Zhu L,Ji L,Liu H
Mon Feb 01 00:00:00 UTC 2021
Thymoquinone Suppresses the Proliferation, Migration and Invasiveness through Regulating ROS, Autophagic Flux and miR-877-5p in Human Bladder Carcinoma Cells.
International journal of biological sciences
Zhou X,Wang F,Wu H,Chen X,Zhang Y,Lin J,Cai Y,Xiang J,He N,Hu Z,Jin X
Tue Feb 22 00:00:00 UTC 2022
Involvement of NADPH oxidase in patulin-induced oxidative damage and cytotoxicity in HEK293 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Han J,Jin C,Zhong Y,Zhu J,Liu Q,Sun D,Feng J,Xia X,Peng X
Thu Apr 01 00:00:00 UTC 2021
Soy isoflavones alleviate polycystic ovary syndrome in rats by regulating NF- κB signaling pathway.
Bioengineered
Ma X,Li X,Ma L,Chen Y,He S
Wed Dec 01 00:00:00 UTC 2021
Black Tea Alleviates Particulate Matter-Induced Lung Injury via the Gut-Lung Axis in Mice.
Journal of agricultural and food chemistry
Zhao Y,Chen X,Shen J,Xu A,Wang Y,Meng Q,Xu P
Wed Dec 22 00:00:00 UTC 2021
Microglia-derived IL-1β promoted neuronal apoptosis through ER stress-mediated signaling pathway PERK/eIF2α/ATF4/CHOP upon arsenic exposure.
Journal of hazardous materials
Liu X,Chen Y,Wang H,Wei Y,Yuan Y,Zhou Q,Fang F,Shi S,Jiang X,Dong Y,Li X
Sun Sep 05 00:00:00 UTC 2021
Effect of luteinizing hormone concentration on transcriptome and subcellular organelle phenotype of ovarian granulosa cells.
Journal of assisted reproduction and genetics
Wan YT,Liu S,Zhao SK,Luo YY,Lv YS,Qu DN,Liu MH,Li Y
Thu Apr 01 00:00:00 UTC 2021
Astrocyte-selective STAT3 knockdown rescues methamphetamine withdrawal-disrupted spatial memory in mice via restoring the astrocytic capacity of glutamate clearance in dCA1.
Glia
Shi P,Li Z,He T,Li N,Xu X,Yu P,Lu X,Nie J,Liu D,Cai Q,Guan Y,Ge F,Wang J,Guan X
Fri Oct 01 00:00:00 UTC 2021
The screening of drug-induced nephrotoxicity using gold nanocluster-based ratiometric fluorescent probes.
Nanoscale
Liu Q,Li J,Liu X,Yuan L,Zhao L,Chang YT,Liu X,Peng J
Sat Aug 28 00:00:00 UTC 2021
PI3K/AKT pathway mediates the antidepressant- and anxiolytic-like roles of hydrogen sulfide in streptozotocin-induced diabetic rats via promoting hippocampal neurogenesis.
Neurotoxicology
Jiang W,Tang YY,Zhu WW,Li C,Zhang P,Li RQ,Chen YJ,Zou W,Tang XQ
Thu Jul 01 00:00:00 UTC 2021
TBC1 domain family member 23 interacts with Ras-related protein Rab-11A to promote poor prognosis of non-small-cell lung cancer via β1-integrin.
Journal of cellular and molecular medicine
Zhang Y,Su H,Wudu M,Ren H,Xu Y,Zhang Q,Jiang J,Wang Q,Jiang X,Zhang B,Liu Z,Zou Z,Qiu X
Wed Sep 01 00:00:00 UTC 2021
The HDAC and PI3K dual inhibitor CUDC-907 synergistically enhances the antileukemic activity of venetoclax in preclinical models of acute myeloid leukemia.
Haematologica
Li X,Su Y,Hege K,Madlambayan G,Edwards H,Knight T,Polin L,Kushner J,Dzinic SH,White K,Yang J,Miller R,Wang G,Zhao L,Wang Y,Lin H,Taub JW,Ge Y
Sat May 01 00:00:00 UTC 2021
Ginsenoside Rg5 Improves Insulin Resistance and Mitochondrial Biogenesis of Liver via Regulation of the Sirt1/PGC-1α Signaling Pathway in db/db Mice.
Journal of agricultural and food chemistry
Zhu Y,Yang H,Deng J,Fan D
Wed Aug 04 00:00:00 UTC 2021
LncRNA MALAT1 promotes breast cancer progression by sponging miR101-3p to mediate mTOR/PKM2 signal transmission.
American journal of translational research
Shao J,Zhang Q,Wang P,Wang Z
Sat Oct 16 00:00:00 UTC 2021
Subunit Vaccine ESAT-6:c-di-AMP Delivered by Intranasal Route Elicits Immune Responses and Protects Against Mycobacterium tuberculosis Infection.
Frontiers in cellular and infection microbiology
Ning H,Zhang W,Kang J,Ding T,Liang X,Lu Y,Guo C,Sun W,Wang H,Bai Y,Shen L
Mon Jul 05 00:00:00 UTC 2021
Fucoxanthin extracted from Laminaria Japonica inhibits metastasis and enhances the sensitivity of lung cancer to Gefitinib.
Journal of ethnopharmacology
Ming JX,Wang ZC,Huang Y,Ohishi H,Wu RJ,Shao Y,Wang H,Qin MY,Wu ZL,Li YY,Chang Zhou S,Chen H,Liu H,Xu R
Sat Jan 30 00:00:00 UTC 2021
Methamphetamine mediates apoptosis of vascular smooth muscle cells via the chop-related endoplasmic reticulum stress pathway.
Toxicology letters
Tan X,Cai D,Chen N,Du S,Qiao D,Yue X,Wang T,Li J,Xie W,Wang H
Sun Oct 10 00:00:00 UTC 2021
Punicalin Attenuates Breast Cancer-Associated Osteolysis by Inhibiting the NF-κB Signaling Pathway of Osteoclasts.
Frontiers in pharmacology
Li T,Jiang G,Hu X,Yang D,Tan T,Gao Z,Chen Z,Xiang C,Li S,Ouyang Z,Guo X
Tue Dec 07 00:00:00 UTC 2021
Berberine Inhibits MDA-MB-231 Cells as an Agonist of G Protein-Coupled Estrogen Receptor 1.
International journal of molecular sciences
Qi M,Liu X,Zhou Y,Wang H,Zhao Y,Ren J,Xiang J
Sun Oct 24 00:00:00 UTC 2021
Novel lncRNA LINC01614 Facilitates Bladder Cancer Proliferation, Migration and Invasion Through the miR-217/RUNX2/Wnt/β-Catenin Axis.
Cancer management and research
Wang Z,Yan H,Cheng D,Xu L,Shen T,Chen Y,Han R,Xue Y
Thu Jun 30 00:00:00 UTC 2022
The SRSF1/circATP5B/miR-185-5p/HOXB5 feedback loop regulates the proliferation of glioma stem cells via the IL6-mediated JAK2/STAT3 signaling pathway.
Journal of experimental & clinical cancer research : CR
Zhao J,Jiang Y,Zhang H,Zhou J,Chen L,Li H,Xu J,Zhang G,Jing Z
Thu Apr 15 00:00:00 UTC 2021
Identification and functional analysis of senescence-associated secretory phenotype of premature senescent hepatocytes induced by hexavalent chromium.
Ecotoxicology and environmental safety
Ma Y,Liang Y,Liang N,Zhang Y,Xiao F
Mon Mar 15 00:00:00 UTC 2021
The combination of G-CSF and AMD3100 mobilizes bone marrow-derived stem cells to protect against cisplatin-induced acute kidney injury in mice.
Stem cell research & therapy
Chen Z,Ren X,Ren R,Wang Y,Shang J
Wed Mar 24 00:00:00 UTC 2021
Essential Role of CRIM1 on Endometrial Receptivity in Goat.
International journal of molecular sciences
Yang D,Liu A,Zhang Y,Nan S,Yin R,Lei Q,Zhu H,Chen J,Han L,Ding M,Ding Y
Tue May 18 00:00:00 UTC 2021
RNF128 suppresses the malignancy of colorectal cancer cells via inhibition of Wnt/β-catenin signaling.
American journal of translational research
Zhu Y,Gan Y,Zou R,Sha H,Lu Y,Zhang Y,Feng J
Tue Jan 18 00:00:00 UTC 2022
Vitamin A and retinoic acid accelerate the attenuation of intestinal adaptability upon feeding induced by high-fat diet in mice.
The Journal of nutritional biochemistry
Hong W,Xu D,Song X,Niu B,Zhuang Z,Lu Y,Lei X,Ma R,Lu C,Sun N,Mao Y,Li X
Mon Nov 01 00:00:00 UTC 2021
Circ-NOLC1 promotes epithelial ovarian cancer tumorigenesis and progression by binding ESRP1 and modulating CDK1 and RhoA expression.
Cell death discovery
Chen S,Wu W,Li QH,Xie BM,Shen F,Du YP,Zong ZH,Wang LL,Wei XQ,Zhao Y
Fri Jan 22 00:00:00 UTC 2021
Targeting Wnt Signaling in the Tumor Immune Microenvironment to Enhancing EpCAM CAR T-Cell therapy.
Frontiers in pharmacology
Li W,Zhou Y,Wu Z,Shi Y,Tian E,Zhu Y,Wang T,Dou W,Meng X,Chen M,Zhai B,Zhu D
Fri Nov 19 00:00:00 UTC 2021
Enhanced response to PD-L1 silencing by modulation of TME via balancing glucose metabolism and robust co-delivery of siRNA/Resveratrol with dual-responsive polyplexes.
Biomaterials
Jia L,Gao Y,Zhou T,Zhao XL,Hu HY,Chen DW,Qiao MX
Thu Apr 01 00:00:00 UTC 2021
Endogenous reverse transcriptase and RNase H-mediated antiviral mechanism in embryonic stem cells.
Cell research
Wu J,Wu C,Xing F,Cao L,Zeng W,Guo L,Li P,Zhong Y,Jiang H,Luo M,Shi G,Bu L,Ji Y,Hou P,Peng H,Huang J,Li C,Guo D
Wed Sep 01 00:00:00 UTC 2021
Pretreatment of Indobufen and Aspirin and their Combinations with Clopidogrel or Ticagrelor Alleviates Inflammasome Mediated Pyroptosis Via Inhibiting NF-κB/NLRP3 Pathway in Ischemic Stroke.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology
Li F,Xu D,Hou K,Gou X,Lv N,Fang W,Li Y
Wed Dec 01 00:00:00 UTC 2021
Combination of ferulic acid, ligustrazine and tetrahydropalmatine inhibits invasion and metastasis through MMP/TIMP signaling in endometriosis.
PeerJ
Tan Y,Zhang C,Zhang Y,Dai X,Wei Q,Wei J,Xu P,Chen Y
Sun Apr 24 00:00:00 UTC 2022
CircFAM114A2 Promotes Cisplatin Sensitivity via miR-222-3p/P27 and miR-146a-5p/P21 Cascades in Urothelial Carcinoma.
Frontiers in oncology
Lv J,Zhou Z,Wang J,Yang X,Yu H,Han J,Feng D,Yuan B,Wu Q,Li P,Lu Q,Yang H
Wed Apr 27 00:00:00 UTC 2022
GRIN2A Variants Associated With Idiopathic Generalized Epilepsies.
Frontiers in molecular neuroscience
Liu XR,Xu XX,Lin SM,Fan CY,Ye TT,Tang B,Shi YW,Su T,Li BM,Yi YH,Luo JH,Liao WP
Tue Nov 02 00:00:00 UTC 2021
Oxymatrine inhibits neuroinflammation byRegulating M1/M2 polarization in N9 microglia through the TLR4/NF-κB pathway.
International immunopharmacology
Wang XL,Chen F,Shi H,Zhang M,Yan L,Pei XY,Peng XD
Mon Nov 01 00:00:00 UTC 2021
BML-111, the lipoxin A4 agonist, modulates VEGF or CoCl2-induced migration, angiogenesis and permeability in tumor-derived endothelial cells.
Immunology letters
Lin L,Wang Q,Xu F,Luo X,Xu J,Yan L,Li Q,Hao H
Mon Feb 01 00:00:00 UTC 2021
Esophageal Cancer Stem-like Cells Resist Ferroptosis-Induced Cell Death by Active Hsp27-GPX4 Pathway.
Biomolecules
Liu CC,Li HH,Lin JH,Chiang MC,Hsu TW,Li AF,Yen DH,Hsu HS,Hung SC
Wed Dec 29 00:00:00 UTC 2021
Honokiol induces paraptosis-like cell death of acute promyelocytic leukemia via mTOR & MAPK signaling pathways activation.
Apoptosis : an international journal on programmed cell death
Liu X,Gu Y,Bian Y,Cai D,Li Y,Zhao Y,Zhang Z,Xue M,Zhang L
Thu Apr 01 00:00:00 UTC 2021
Inhibition of Growth of Colon Tumors and Proliferation of HT-29 Cells by Warburgia ugandensis Extract through Mediating G0/G1 Cell Cycle Arrest, Cell Apoptosis, and Intracellular ROS Generation.
Oxidative medicine and cellular longevity
Zhang Y,Chen G,Zhuang X,Guo M
Fri Mar 04 00:00:00 UTC 2022
Resveratrol induces human colorectal cancer cell apoptosis by activating the mitochondrial pathway via increasing reactive oxygen species.
Molecular medicine reports
Fu Y,Ye Y,Zhu G,Xu Y,Sun J,Wu H,Feng F,Wen Z,Jiang S,Li Y,Zhang Q
Mon Mar 01 00:00:00 UTC 2021
Hippocampal CA1 theta burst-induced LTP from weaning to adulthood: Cellular and molecular mechanisms in young male rats revisited.
The European journal of neuroscience
Rodrigues NC,Silva-Cruz A,Caulino-Rocha A,Bento-Oliveira A,Alexandre Ribeiro J,Cunha-Reis D
Sun Aug 01 00:00:00 UTC 2021
Danthron ameliorates obesity and MAFLD through activating the interplay between PPARα/RXRα heterodimer and adiponectin receptor 2.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ma C,Wang Z,Xia R,Wei L,Zhang C,Zhang J,Zhao L,Wu H,Kang L,Yang S
Sat May 01 00:00:00 UTC 2021
BLM helicase inhibition synergizes with PARP inhibition to improve the radiosensitivity of olaparib resistant non-small cell lung cancer cells by inhibiting homologous recombination repair.
Cancer biology & medicine
Kong Y,Xu C,Sun X,Sun H,Zhao X,He N,Ji K,Wang Q,Du L,Wang J,Zhang M,Liu Y,Wang Y,Liu Q
Wed Dec 01 00:00:00 UTC 2021
Human Cytomegalovirus Uses a Host Stress Response To Balance the Elongation of Saturated/Monounsaturated and Polyunsaturated Very-Long-Chain Fatty Acids.
mBio
Xi Y,Lindenmayer L,Kline I,von Einem J,Purdy JG
Tue May 04 00:00:00 UTC 2021
Protective effect of naringenin against cadmium-induced testicular toxicity in male SD rats.
Journal of inorganic biochemistry
Wang J,Zhu H,Lin S,Wang K,Wang H,Liu Z
Fri Jan 01 00:00:00 UTC 2021
Electroacupuncture Preconditioning Reduces Oxidative Stress in the Acute Phase of Cerebral Ischemia-Reperfusion in Rats by Regulating Iron Metabolism Pathways.
Evidence-based complementary and alternative medicine : eCAM
Liang R,Tang Q,Song W,Zhang M,Teng L,Kang Y,Zhu L
Thu Apr 28 00:00:00 UTC 2022
Lipoxin A4 protects against paraquat‑induced acute lung injury by inhibiting the TLR4/MyD88‑mediated activation of the NF‑κB and PI3K/AKT pathways.
International journal of molecular medicine
Li Y,Wang N,Ma Z,Wang Y,Yuan Y,Zhong Z,Hong Y,Zhao M
Sat May 01 00:00:00 UTC 2021
miR-126 Controls the Apoptosis and Proliferation of Immature Porcine Sertoli Cells by Targeting the PIK3R2 Gene through the PI3K/AKT Signaling Pathway.
Animals : an open access journal from MDPI
Tang X,Chen Y,Luo H,Bian Q,Weng B,Yang A,Chu D,Ran M,Chen B
Fri Jul 30 00:00:00 UTC 2021
Obacunone Attenuates Liver Fibrosis with Enhancing Anti-Oxidant Effects of GPx-4 and Inhibition of EMT.
Molecules (Basel, Switzerland)
Bai Y,Wang W,Wang L,Ma L,Zhai D,Wang F,Shi R,Liu C,Xu Q,Chen G,Lu Z
Sat Jan 09 00:00:00 UTC 2021
Osteopontin N-Terminal Function in an Abdominal Aortic Aneurysm From Apolipoprotein E-Deficient Mice.
Frontiers in cell and developmental biology
Liu H,Zhang Y,Song W,Sun Y,Jiang Y
Tue Aug 31 00:00:00 UTC 2021
Down-regulation of EPB41L4A-AS1 mediated the brain aging and neurodegenerative diseases via damaging synthesis of NAD+ and ATP.
Cell & bioscience
Yang T,Wang Y,Liao W,Zhang S,Wang S,Xu N,Xie W,Luo C,Wang Y,Wang Z,Zhang Y
Wed Nov 10 00:00:00 UTC 2021
Hypoxia-induced FOXO4/LDHA axis modulates gastric cancer cell glycolysis and progression.
Clinical and translational medicine
Wang XH,Jiang ZH,Yang HM,Zhang Y,Xu LH
Fri Jan 01 00:00:00 UTC 2021
MMP11 promotes the proliferation and progression of breast cancer through stabilizing Smad2 protein.
Oncology reports
Zhuang Y,Li X,Zhan P,Pi G,Wen G
Thu Apr 01 00:00:00 UTC 2021
HORMAD1 promotes docetaxel resistance in triple negative breast cancer by enhancing DNA damage tolerance.
Oncology reports
Zong B,Sun L,Peng Y,Wang Y,Yu Y,Lei J,Zhang Y,Guo S,Li K,Liu S
Thu Jul 01 00:00:00 UTC 2021
The effects of Omarigliptin on promoting osteoblastic differentiation.
Bioengineered
Liao F,Hu X,Chen R
Wed Dec 01 00:00:00 UTC 2021
Dihydroartemisinin suppresses proliferation, migration, the Wnt/β-catenin pathway and EMT via TNKS in gastric cancer.
Oncology letters
Ma Y,Zhang P,Zhang Q,Wang X,Miao Q,Lyu X,Cui B,Ma H
Fri Oct 01 00:00:00 UTC 2021
Ferrostatin-1 alleviated TNBS induced colitis via the inhibition of ferroptosis.
Biochemical and biophysical research communications
Xu J,Liu S,Cui Z,Wang X,Ning T,Wang T,Zhang N,Xie S,Min L,Zhang S,Liang C,Zhu S
Fri Oct 08 00:00:00 UTC 2021
Augmenter of liver regeneration ameliorates ischemia-reperfusion injury in steatotic liver via inhibition of the TLR4/NF-κB pathway.
Experimental and therapeutic medicine
Weng J,Wang X,Xu B,Li W
Sun Aug 01 00:00:00 UTC 2021
Changes in the prefrontal cortex after the hippocampus was injected with Aβ25-35 via the P35/P25-CDK5-Tau hyperphosphorylation signaling pathway.
Neuroscience letters
Wang Y,Sheng H,Zhao J,Guo L,Liu J,Xu J,Liu Q,Huang J,Jiang R,Gan S,Qiu G,Lu W,Xu S,Zhu S
Sun Jan 10 00:00:00 UTC 2021
Triptolide regulates oxidative stress and inflammation leading to hepatotoxicity via inducing CYP2E1.
Human & experimental toxicology
Jiang HY,Bao YN,Lin FM,Jin Y
Wed Dec 01 00:00:00 UTC 2021
Gamma Radiation Induce Inflammasome Signaling and Pyroptosis in Microvascular Endothelial Cells.
Journal of inflammation research
Smith AO,Ju W,Adzraku SY,Wenyi L,Yuting C,Qiao J,Xu K,Zeng L
Mon Apr 25 00:00:00 UTC 2022
Brd4 Regulates the Homeostasis of CD8+ T-Lymphocytes and Their Proliferation in Response to Antigen Stimulation.
Frontiers in immunology
Peng Z,Zhang Y,Ma X,Zhou M,Wu S,Song Z,Yuan Y,Chen Y,Li Y,Wang G,Huang F,Qiao Y,Xia B,Liu W,Liu J,Zhang X,He X,Pan T,Xu H,Zhang H
Mon Dec 27 00:00:00 UTC 2021
Novel clerodane-type diterpenoid Cintelactone A suppresses lipopolysaccharide -induced inflammation by promoting ubiquitination, proteasomal degradation of TRAF6.
Pharmacological research
Di Q,Zhao X,Zhang R,Ma X,Liang X,Li X,Gao J,Tang H,Chen W,Xiao W
Mon Feb 01 00:00:00 UTC 2021
Inhibition of TNFAIP1 ameliorates the oxidative stress and inflammatory injury in myocardial ischemia/reperfusion injury through modulation of Akt/GSK-3β/Nrf2 pathway.
International immunopharmacology
Wen L,Yang QH,Ma XL,Li T,Xiao S,Sun CF
Fri Oct 01 00:00:00 UTC 2021
Reduced DAPK1 Expression Promotes Stem Cell-Like Characteristics of Prostate Cancer Cells by Activating ZEB1 via Hippo/YAP Signaling Pathway.
Stem cells and development
Nong S,Wei Z,Wang Z,Ma L,Guan Y,Ni J
Wed Sep 15 00:00:00 UTC 2021
EPZ015666, a selective protein arginine methyltransferase 5 (PRMT5) inhibitor with an antitumour effect in retinoblastoma.
Experimental eye research
Liu X,He J,Mao L,Zhang Y,Cui W,Duan S,Jiang A,Gao Y,Sang Y,Huang G
Fri Jan 01 00:00:00 UTC 2021
Nano chitosan-zinc complex improves the growth performance and antioxidant capacity of the small intestine in weaned piglets.
The British journal of nutrition
Zhang M,Hou G,Hu P,Feng D,Wang J,Zhu W
Tue Sep 28 00:00:00 UTC 2021
Limonin ameliorates dextran sulfate sodium-induced chronic colitis in mice by inhibiting PERK-ATF4-CHOP pathway of ER stress and NF-κB signaling.
International immunopharmacology
Song C,Chen J,Li X,Yang R,Cao X,Zhou L,Zhou Y,Ying H,Zhang Q,Sun Y
Fri Jan 01 00:00:00 UTC 2021
Aged Ripe Pu-erh Tea Reduced Oxidative Stress-Mediated Inflammation in Dextran Sulfate Sodium-Induced Colitis Mice by Regulating Intestinal Microbes.
Journal of agricultural and food chemistry
Hu S,Li S,Liu Y,Sun K,Luo L,Zeng L
Wed Sep 15 00:00:00 UTC 2021
Sepsis induces muscle atrophy by inhibiting proliferation and promoting apoptosis via PLK1-AKT signalling.
Journal of cellular and molecular medicine
Cao YY,Wang Z,Yu T,Zhang Y,Wang ZH,Lu ZM,Lu WH,Yu JB
Fri Oct 01 00:00:00 UTC 2021
Cathepsin K deficiency promotes alveolar bone regeneration by promoting jaw bone marrow mesenchymal stem cells proliferation and differentiation via glycolysis pathway.
Cell proliferation
Zhang W,Dong Z,Li D,Li B,Liu Y,Zheng X,Liu H,Zhou H,Hu K,Xue Y
Thu Jul 01 00:00:00 UTC 2021
ING5 Inhibits Migration and Invasion of Esophageal Cancer Cells by Downregulating the IL-6/CXCL12 Signaling Pathway.
Technology in cancer research & treatment
Wang Y,Tan J,Li J,Chen H,Wang W
Mon Jan 24 00:00:00 UTC 2022
RhoA/ROCK-YAP/TAZ Axis Regulates the Fibrotic Activity in Dexamethasone-Treated Human Trabecular Meshwork Cells.
Frontiers in molecular biosciences
Liu Z,Li S,Qian X,Li L,Zhang H,Liu Z
Sat Sep 25 00:00:00 UTC 2021
ARHGEF3 Associated with Invasion, Metastasis, and Proliferation in Human Osteosarcoma.
BioMed research international
Gong J,Tang W,Lv B,Zhang S,Fan T,Gao G,Chen D,Liu Y
Thu Sep 23 00:00:00 UTC 2021
Toxicity evaluation of mesoporous silica particles Santa Barbara No. 15 amorphous in human umbilical vein endothelial cells: influence of particle morphology.
Journal of applied toxicology : JAT
Teng W,Yang Z,Wang S,Xiong D,Chen Y,Wu Z
Wed Sep 01 00:00:00 UTC 2021
DHAV-1 Blocks the Signaling Pathway Upstream of Type I Interferon by Inhibiting the Interferon Regulatory Factor 7 Protein.
Frontiers in microbiology
Lai Y,Xia X,Cheng A,Wang M,Ou X,Mao S,Sun D,Zhang S,Yang Q,Wu Y,Zhu D,Jia R,Chen S,Liu M,Zhao XX,Huang J,Gao Q,Tian B,Liu Y,Yu Y,Zhang L,Pan L
Tue Dec 07 00:00:00 UTC 2021
Cell membrane-encapsulated magnetic nanoparticles for enhancing natural killer cell-mediated cancer immunotherapy.
Nanomedicine : nanotechnology, biology, and medicine
Wu D,Shou X,Zhang Y,Li Z,Wu G,Wu D,Wu J,Shi S,Wang S
Mon Feb 01 00:00:00 UTC 2021
A genome-wide CRISPR screen identifies host factors that regulate SARS-CoV-2 entry.
Nature communications
Zhu Y,Feng F,Hu G,Wang Y,Yu Y,Zhu Y,Xu W,Cai X,Sun Z,Han W,Ye R,Qu D,Ding Q,Huang X,Chen H,Xu W,Xie Y,Cai Q,Yuan Z,Zhang R
Thu Feb 11 00:00:00 UTC 2021
Glia Maturation Factor Beta as a Novel Biomarker and Therapeutic Target for Hepatocellular Carcinoma.
Frontiers in oncology
Sun W,Hu C,Wang T,Wang J,Zhang J,Gao F,Ou Q,Tian H,Jin C,Xu J,Zhang J,Xu GT,Lu L
Sat Nov 20 00:00:00 UTC 2021
Comprehensive Analysis of the Potential Immune-Related Biomarker Transporter Associated With Antigen Processing 1 That Inhibits Metastasis and Invasion of Ovarian Cancer Cells.
Frontiers in molecular biosciences
Li X,Zeng S,Ding Y,Nie Y,Yang M
Mon Sep 19 00:00:00 UTC 2022
Remarkable inhibition effects of afatinib alone or combining with paclitaxel in esophageal squamous cell carcinoma.
Journal of gastroenterology and hepatology
Yang LY,Cheng ZJ,Liu Z,Wang D,Zhang N,Fan ZL,Cai HQ,Zhang Y,Cai Y,Xu X,Wang JH,Du GH,Hao JJ,Wang MR
Wed Sep 01 00:00:00 UTC 2021
The long non-coding RNA PFI protects against pulmonary fibrosis by interacting with splicing regulator SRSF1.
Cell death and differentiation
Sun J,Jin T,Su W,Guo Y,Niu Z,Guo J,Li L,Wang J,Ma L,Yu T,Li X,Zhou Y,Shan H,Liang H
Fri Oct 01 00:00:00 UTC 2021
Estrogen Receptor α Mediates Doxorubicin Sensitivity in Breast Cancer Cells by Regulating E-Cadherin.
Frontiers in cell and developmental biology
Wan X,Hou J,Liu S,Zhang Y,Li W,Zhang Y,Ding Y
Tue Feb 23 00:00:00 UTC 2021
Study on Protection of Human Umbilical Vein Endothelial Cells from Amiodarone-Induced Damage by Intermedin through Activation of Wnt/β-Catenin Signaling Pathway.
Oxidative medicine and cellular longevity
Wang Y,Wang J,Yang J,Kang J,Xue F,Chang S,Ji H,Zang H,Zhou X,Wang G,Fan W,Yan X,Guo J,Ren X,Tian J
Wed Jan 19 00:00:00 UTC 2022
Rational design and synthesis of 6-aryl-6H-benzo[c]chromenes as non-steroidal progesterone receptor antagonists for use against cancers.
Bioorganic & medicinal chemistry
Qin J,Qu S,Zhu K,Cheng Y,Pan G,Jing W,Liu X,Sun X,Liu L
Mon Feb 15 00:00:00 UTC 2021
QiShenYiQi Pill Improves Myocardial Hypertrophy Caused by Pressure Overload in Rats.
Evidence-based complementary and alternative medicine : eCAM
Lv S,Wang Q,Wu M,Li M,Wang X,Xu L,Zhang J
Tue Jul 06 00:00:00 UTC 2021
Bioengineering CXCR4-overexpressing cell membrane functionalized ROS-responsive nanotherapeutics for targeting cerebral ischemia-reperfusion injury.
Theranostics
Luo L,Zang G,Liu B,Qin X,Zhang Y,Chen Y,Zhang H,Wu W,Wang G
Tue Aug 17 00:00:00 UTC 2021
The Panda-Derived Lactobacillus plantarum G201683 Alleviates the Inflammatory Response in DSS-Induced Panda Microbiota-Associated Mice.
Frontiers in immunology
Zhou Y,Duan L,Zeng Y,Niu L,Pu Y,Jacobs JP,Chang C,Wang J,Khalique A,Pan K,Fang J,Jing B,Zeng D,Ni X
Mon Feb 07 00:00:00 UTC 2022
Medioresinol as a novel PGC-1α activator prevents pyroptosis of endothelial cells in ischemic stroke through PPARα-GOT1 axis.
Pharmacological research
Wang Y,Guan X,Gao CL,Ruan W,Zhao S,Kai G,Li F,Pang T
Thu Jul 01 00:00:00 UTC 2021
Change on apoptosis, autophagy and mitochondria of the Harderian gland in Cricetulus barabensis during age.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology
Kong XT,Wang Z,Mou JJ,Li CS,Xue HL,Wu M,Chen L,Xu JH,Xu LX
Mon Jun 28 00:00:00 UTC 2021
Glucagon-like peptide-1 (GLP-1) improved diabetic lung fibrosis via AMPK and microRNA-27a (miR-27a).
Annals of translational medicine
Liu J,Li X,Lu S,Zheng X,Zhang X,Zhao W
Mon Mar 01 00:00:00 UTC 2021
L-type amino acid transporter 1 is associated with chemoresistance in breast cancer via the promotion of amino acid metabolism.
Scientific reports
Sato M,Harada-Shoji N,Toyohara T,Soga T,Itoh M,Miyashita M,Tada H,Amari M,Anzai N,Furumoto S,Abe T,Suzuki T,Ishida T,Sasano H
Tue Jan 12 00:00:00 UTC 2021
Different effects of high-fat diets rich in different oils on lipids metabolism, oxidative stress and gut microbiota.
Food research international (Ottawa, Ont.)
Liu H,Zhu H,Xia H,Yang X,Yang L,Wang S,Wen J,Sun G
Mon Mar 01 00:00:00 UTC 2021
ActivinA activates Notch1-Shh signaling to regulate proliferation in C2C12 skeletal muscle cells.
Molecular and cellular endocrinology
Ma L,Li C,Lian S,Xu B,Yuan J,Lu J,Yang H,Guo J,Ji H
Fri Jan 01 00:00:00 UTC 2021
Different concentrations of C5a affect human dental pulp mesenchymal stem cells differentiation.
BMC oral health
Liu J,Wei X,Hu J,Tan X,Kang X,Gao L,Li N,Shi X,Yuan M,Hu W,Liu M
Fri Sep 24 00:00:00 UTC 2021
Formaldehyde induces ferroptosis in hippocampal neuronal cells by upregulation of the Warburg effect.
Toxicology
Li XN,Yang SQ,Li M,Li XS,Tian Q,Xiao F,Tang YY,Kang X,Wang CY,Zou W,Zhang P,Tang XQ
Sat Jan 30 00:00:00 UTC 2021
Raf1 interacts with OIP5 to participate in oxaliplatin-induced neuropathic pain.
Life sciences
Yu W,Zheng Z,Wei W,Li L,Zhang Y,Sun Y,Cao J,Zang W,Shao J
Wed Sep 15 00:00:00 UTC 2021
Menthol Targeting AMPK Alleviates the Inflammatory Response of Bovine Mammary Epithelial Cells and Restores the Synthesis of Milk Fat and Milk Protein.
Frontiers in immunology
Liu S,Guo W,Jia Y,Ye B,Liu S,Fu S,Liu J,Hu G
Mon Feb 14 00:00:00 UTC 2022
Yap1-2 Isoform Is the Primary Mediator in TGF-β1 Induced EMT in Pancreatic Cancer.
Frontiers in oncology
Gao C,Quan MY,Chen QJ,Yang R,Wu Y,Liu JY,Lin ZY,Li X,Cai JT,Jiang TF,Xu L,Mossahebi-Mohammadi M,Guo Q,Zhang JS
Tue Jun 08 00:00:00 UTC 2021
Anterior Gradient Protein 2 Promotes Mucosal Repair in Pediatric Ulcerative Colitis.
BioMed research international
Ye X,Wu J,Li J,Wang H
Mon Sep 20 00:00:00 UTC 2021
Differentially expressed genes induced by β-caryophyllene in a rat model of cerebral ischemia-reperfusion injury.
Life sciences
Liu S,Liu J,Wang Y,Deng L,Chen S,Wang X,Zuo T,Hu Q,Rao J,Wang Q,Dong Z
Sat May 15 00:00:00 UTC 2021
T-2 Toxin Induces Ferroptosis by Increasing Lipid Reactive Oxygen Species (ROS) and Downregulating Solute Carrier Family 7 Member 11 (SLC7A11).
Journal of agricultural and food chemistry
Wang G,Qin S,Zheng Y,Xia C,Zhang P,Zhang L,Yao J,Yi Y,Deng L
Wed Dec 29 00:00:00 UTC 2021
The novel ALK inhibitor ZX-29 induces apoptosis through inhibiting ALK and inducing ROS-mediated endoplasmic reticulum stress in Karpas299 cells.
Journal of biochemical and molecular toxicology
Zhou X,Zhang X,Wu Z,Xu X,Guo M,Zhai X,Zuo D,Wu Y
Mon Mar 01 00:00:00 UTC 2021
Bergenin alleviates H2 O2 -induced oxidative stress and apoptosis in nucleus pulposus cells: Involvement of the PPAR-γ/NF-κB pathway.
Environmental toxicology
Zhang G,Wang H,Zhang Q,Zhao Z,Zhu W,Zuo X
Wed Dec 01 00:00:00 UTC 2021
Curcumin-Alleviated Osteoarthritic Progression in Rats Fed a High-Fat Diet by Inhibiting Apoptosis and Activating Autophagy via Modulation of MicroRNA-34a.
Journal of inflammation research
Yao J,Liu X,Sun Y,Dong X,Liu L,Gu H
Sat Apr 23 00:00:00 UTC 2022
Dendrobium nobile Lindl alkaloid attenuates 6-OHDA-induced dopamine neurotoxicity.
Biotechnology and applied biochemistry
Li DD,Wang GQ,Wu Q,Shi JS,Zhang F
Wed Dec 01 00:00:00 UTC 2021
Maiwei Yangfei decoction prevents bleomycin-induced pulmonary fibrosis in mice.
Experimental and therapeutic medicine
Xu Y,Peng W,Han D,Feng F,Wang Z,Gu C,Zhou X,Wu Q
Mon Nov 01 00:00:00 UTC 2021
LncRNA H19 Upregulation Participates in the Response of Glioma Cells to Radiation.
BioMed research international
Kuang Y,Bing Z,Jin X,Li Q
Mon Oct 18 00:00:00 UTC 2021
Study on the protective effect and mechanism of Dicliptera chinensis (L.) Juss (Acanthaceae) polysaccharide on immune liver injury induced by LPS.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Xu Q,Xu J,Zhang K,Zhong M,Cao H,Wei R,Jin L,Gao Y
Mon Feb 01 00:00:00 UTC 2021
Resveratrol-mediated neurorestoration after cerebral ischemic injury - Sonic Hedgehog signaling pathway.
Life sciences
Yu P,Wang L,Tang F,Guo S,Liao H,Fan C,Yang Q
Wed Sep 01 00:00:00 UTC 2021
Activation of α7nAChR Protects Against Gastric Inflammation and Dysmotility in Parkinson's Disease Rats.
Frontiers in pharmacology
Zhou L,Zheng LF,Zhang XL,Wang ZY,Yao YS,Xiu XL,Liu CZ,Zhang Y,Feng XY,Zhu JX
Sat Dec 11 00:00:00 UTC 2021
33-kDa ANXA3 isoform contributes to hepatocarcinogenesis via modulating ERK, PI3K/Akt-HIF and intrinsic apoptosis pathways.
Journal of advanced research
Guo C,Li N,Dong C,Wang L,Li Z,Liu Q,Ma Q,Greenaway FT,Tian Y,Hao L,Liu S,Sun MZ
Sat May 01 00:00:00 UTC 2021
Indoleamine-2,3-Dioxygenase Activates Wnt/β-Catenin Inducing Kidney Fibrosis after Acute Kidney Injury.
Gerontology
Pan B,Zhang H,Hong Y,Ma M,Wan X,Cao C
Tue Nov 16 00:00:00 UTC 2021
Ubash3b promotes TPA-mediated suppression of leukemogenesis through accelerated downregulation of PKCδ protein.
Biochimie
Yao Y,Liu W,Gajendran B,Wang C,Zacksenhaus E,Sample KM,Varier KM,Hao X,Ben-David Y
Sat May 01 00:00:00 UTC 2021
A deafness-associated tRNA mutation caused pleiotropic effects on the m1G37 modification, processing, stability and aminoacylation of tRNAIle and mitochondrial translation.
Nucleic acids research
Meng F,Zhou M,Xiao Y,Mao X,Zheng J,Lin J,Lin T,Ye Z,Cang X,Fu Y,Wang M,Guan MX
Mon Jan 25 00:00:00 UTC 2021
p53 Promoted Ferroptosis in Ovarian Cancer Cells Treated with Human Serum Incubated-Superparamagnetic Iron Oxides.
International journal of nanomedicine
Zhang Y,Xia M,Zhou Z,Hu X,Wang J,Zhang M,Li Y,Sun L,Chen F,Yu H
Thu Jan 28 00:00:00 UTC 2021
Transient Receptor Potential Melastatin 7 Promotes Vascular Adventitial Fibroblasts Phenotypic Transformation and Inflammatory Reaction Induced by Mechanical Stretching Stress via p38 MAPK/JNK Pathway.
Journal of vascular research
Liu J,Chen L,Huang J,Guo S,Zhu D,Gao P
Mon Dec 20 00:00:00 UTC 2021
MCU That Is Transcriptionally Regulated by Nrf2 Augments Malignant Biological Behaviors in Oral Squamous Cell Carcinoma Cells.
BioMed research international
Wu R,Zuo W,Xu X,Bi L,Zhang C,Chen H,Liu H
Mon Oct 25 00:00:00 UTC 2021
Induction of autophagy by Beclin-1 in granulosa cells contributes to follicular progesterone elevation in ovarian endometriosis.
Translational research : the journal of laboratory and clinical medicine
Ding Y,Zhu Q,He Y,Lu Y,Wang Y,Qi J,Wu H,Xu R,Li J,Li X,Sun Y
Fri Jan 01 00:00:00 UTC 2021
Programmed cell death 5 improves skeletal muscle insulin resistance by inhibiting IRS-1 ubiquitination through stabilization of MDM2.
Life sciences
Li B,Ye J,Liu R,Weng L,Cao Y,Jia S,Xu C,Liu Y,Yan S,Zheng M
Mon Nov 15 00:00:00 UTC 2021
Metformin-Induced MicroRNA-34a-3p Downregulation Alleviates Senescence in Human Dental Pulp Stem Cells by Targeting CAB39 through the AMPK/mTOR Signaling Pathway.
Stem cells international
Zhang S,Zhang R,Qiao P,Ma X,Lu R,Wang F,Li C,E L,Liu H
Sat Jan 30 00:00:00 UTC 2021
TRIM15 promotes the invasion and metastasis of pancreatic cancer cells by mediating APOA1 ubiquitination and degradation.
Biochimica et biophysica acta. Molecular basis of disease
Sun Y,Ren D,Yang C,Yang W,Zhao J,Zhou Y,Jin X,Wu H
Mon Nov 01 00:00:00 UTC 2021
Geniposide Ameliorates Liver Fibrosis Through Reducing Oxidative Stress and Inflammatory Respose, Inhibiting Apoptosis and Modulating Overall Metabolism.
Frontiers in pharmacology
Yang L,Bi L,Jin L,Wang Y,Li Y,Li Z,He W,Cui H,Miao J,Wang L
Wed Dec 15 00:00:00 UTC 2021
Carbohydrate response element-binding protein regulates lipid metabolism via mTOR complex1 in diabetic nephropathy.
Journal of cellular physiology
Chen N,Mu L,Yang Z,Du C,Wu M,Song S,Yuan C,Shi Y
Fri Jan 01 00:00:00 UTC 2021
CRSP8 promotes thyroid cancer progression by antagonizing IKKα-induced cell differentiation.
Cell death and differentiation
Liao Y,Hua Y,Li Y,Zhang C,Yu W,Guo P,Zou K,Li W,Sun Y,Wang R,Zuo Y,Sui S,Tian C,Hao J,Chen M,Hu S,Chen M,Long Q,Wang X,Zou L,Xie F,Guo W,Deng W
Thu Apr 01 00:00:00 UTC 2021
Neuroprotective Effects of Oxymatrine on PI3K/Akt/mTOR Pathway After Hypoxic-Ischemic Brain Damage in Neonatal Rats.
Frontiers in pharmacology
Wei W,Lu M,Lan XB,Liu N,Su WK,Dushkin AV,Yu JQ
Sun May 02 00:00:00 UTC 2021
Osteogenic differentiation and calcification of human aortic smooth muscle cells is induced by the RCN2/STAT3/miR-155-5p feedback loop.
Vascular pharmacology
Zhao J,Liu Z,Chang Z
Mon Feb 01 00:00:00 UTC 2021
The Ghrelin/Growth Hormone Secretagogue Receptor System Is Involved in the Rapid and Sustained Antidepressant-Like Effect of Paeoniflorin.
Frontiers in neuroscience
Zhang Y,Zhu MZ,Qin XH,Zeng YN,Zhu XH
Sat Mar 06 00:00:00 UTC 2021
Regulation of survivin protein stability by USP35 is evolutionarily conserved.
Biochemical and biophysical research communications
Wang W,Lin H,Zheng E,Hou Z,Liu Y,Huang W,Chen D,Feng J,Li J,Li L
Fri Oct 15 00:00:00 UTC 2021
Inhibition of ER stress attenuates kidney injury and apoptosis induced by 3-MCPD via regulating mitochondrial fission/fusion and Ca2+ homeostasis.
Cell biology and toxicology
Zhong Y,Jin C,Han J,Zhu J,Liu Q,Sun D,Xia X,Peng X
Fri Oct 01 00:00:00 UTC 2021
Hypoxia Activates Notch4 via ERK/JNK/P38 MAPK Signaling Pathways to Promote Lung Adenocarcinoma Progression and Metastasis.
Frontiers in cell and developmental biology
Li X,Cao X,Zhao H,Guo M,Fang X,Li K,Qin L,He Y,Liu X
Fri Jan 07 00:00:00 UTC 2022
ARRB1 Drives Gallbladder Cancer Progression by Facilitating TAK1/MAPK Signaling Activation.
Journal of Cancer
Zhang X,Kong Z,Xu X,Yun X,Chao J,Ding D,Li T,Gao Y,Guan N,Zhu C,Qin X
Wed Mar 24 00:00:00 UTC 2021
Deciphering the autophagy regulatory network via single-cell transcriptome analysis reveals a requirement for autophagy homeostasis in spermatogenesis.
Theranostics
Wang M,Xu Y,Zhang Y,Chen Y,Chang G,An G,Yang X,Zheng C,Zhao J,Liu Z,Wang D,Miao K,Rao S,Dai M,Wang D,Zhao XY
Mon Aug 02 00:00:00 UTC 2021
Quercetin hinders microglial activation to alleviate neurotoxicity via the interplay between NLRP3 inflammasome and mitophagy.
Redox biology
Han X,Xu T,Fang Q,Zhang H,Yue L,Hu G,Sun L
Sun Aug 01 00:00:00 UTC 2021
MicroRNA-628-5p Facilitates Enterovirus 71 Infection by Suppressing TRAF3 Signaling.
Cellular & molecular immunology
Li D,Chen S,Zhang W,Zhang C,Sun T,Du Y,Ding R,Gao Y,Jin Y,Duan G
Sat May 01 00:00:00 UTC 2021
Irisin Contributes to Neuroprotection by Promoting Mitochondrial Biogenesis After Experimental Subarachnoid Hemorrhage.
Frontiers in aging neuroscience
Tu T,Yin S,Pang J,Zhang X,Zhang L,Zhang Y,Xie Y,Guo K,Chen L,Peng J,Jiang Y
Tue Feb 23 00:00:00 UTC 2021
SPT6 recruits SND1 to co-activate human telomerase reverse transcriptase to promote colon cancer progression.
Molecular oncology
Diao C,Guo P,Yang W,Sun Y,Liao Y,Yan Y,Zhao A,Cai X,Hao J,Hu S,Yu W,Chen M,Wang R,Li W,Zuo Y,Pan J,Hua C,Lu X,Fan W,Zheng Z,Deng W,Luo G,Guo W
Thu Apr 01 00:00:00 UTC 2021
Hypercholesterolemia risk associated Abca6 does not regulate lipoprotein metabolism in mice or hamster.
Biochimica et biophysica acta. Molecular and cell biology of lipids
He B,Kang S,Chen Z,Liu X,Wang J,Li X,Liu X,Zheng L,Luo M,Wang Y
Mon Nov 01 00:00:00 UTC 2021
MicroRNA-122 contributes to lipopolysaccharide-induced acute kidney injury via down-regulating the vitamin D receptor in the kidney.
European journal of clinical investigation
He J,Du J,Yi B,Wang J,Zhang H,Li YC,Sun J
Sun Aug 01 00:00:00 UTC 2021
Type 1 interferon aggravates lipopolysaccharide-induced sepsis through upregulating Caspase-11 and Gasdermin D.
Journal of physiology and biochemistry
Wang Y,Zhang X,Guo Y,Li X,Guo G,Niu Z,Zhang J
Mon Feb 01 00:00:00 UTC 2021
Targeting the p300/NONO axis sensitizes melanoma cells to BRAF inhibitors.
Oncogene
Zhang F,Tang X,Fan S,Liu X,Sun J,Ju C,Liang Y,Liu R,Zhou R,Yu B,Zhang C,Zhang Z,Kang T,Huang G,Lv XB
Tue Jun 01 00:00:00 UTC 2021
Screening and Identification of Differential Ovarian Proteins before and after Induced Ovulation via Seminal Plasma in Bactrian Camels.
Animals : an open access journal from MDPI
Wang Q,Zhang Q,Li Y,Zhao X,Zhang Y
Thu Dec 09 00:00:00 UTC 2021
Guanosine and uridine alleviate airway inflammation via inhibition of the MAPK and NF-κB signals in OVA-induced asthmatic mice.
Pulmonary pharmacology & therapeutics
Luo Y,Chen H,Huang R,Wu Q,Li Y,He Y
Sun Aug 01 00:00:00 UTC 2021
Chlorogenic Acid Improves NAFLD by Regulating gut Microbiota and GLP-1.
Frontiers in pharmacology
Shi A,Li T,Zheng Y,Song Y,Wang H,Wang N,Dong L,Shi H
Tue Jul 20 00:00:00 UTC 2021
Effects of total flavonoids from Eucommia ulmoides Oliv. leaves on polycystic ovary syndrome with insulin resistance model rats induced by letrozole combined with a high-fat diet.
Journal of ethnopharmacology
Peng MF,Tian S,Song YG,Li CX,Miao MS,Ren Z,Li M
Sat Jun 12 00:00:00 UTC 2021
Low EGR1 expression predicts poor prognosis in clear cell renal cell carcinoma.
Pathology, research and practice
Zhang ZY,Zhang SL,Chen HL,Mao YQ,Kong CY,Li ZM,Wang LS,Ma M,Han B
Wed Dec 01 00:00:00 UTC 2021
Non-canonical activation of the ER stress sensor ATF6 by Legionella pneumophila effectors.
Life science alliance
Ibe NU,Subramanian A,Mukherjee S
Wed Dec 01 00:00:00 UTC 2021
Blockade of GITRL/GITR signaling pathway attenuates house dust mite-induced allergic asthma in mice through inhibition of MAPKs and NF-κB signaling.
Molecular immunology
Wang Y,Liu B,Niu C,Zou W,Yang L,Wang T,Tian D,Luo Z,Dai J,Li Q,Liu E,Gong C,Fu Z,Ding F
Wed Sep 01 00:00:00 UTC 2021
Dietary seaweed-derived polysaccharides improve growth performance of weaned pigs through maintaining intestinal barrier function and modulating gut microbial populations.
Journal of animal science and biotechnology
Zou T,Yang J,Guo X,He Q,Wang Z,You J
Wed Mar 10 00:00:00 UTC 2021
MSH6 Aggravates the Hypoxic Microenvironment via Regulating HIF1A to Promote the Metastasis of Glioblastoma Multiforme.
DNA and cell biology
Shi Y,Jiang J,Cui Y,Chen Y,Dong T,An H,Liu P
Fri Jan 01 00:00:00 UTC 2021
Long non-coding RNA CRNDE exacerbates NPC advancement mediated by the miR-545-5p/CCND2 axis.
Cancer cell international
Ge S,Jiang C,Li M,Cheng Z,Feng X
Sat Dec 04 00:00:00 UTC 2021
ZEB1 represses biogenesis of circ-DOCK5 to facilitate metastasis in esophageal squamous cell carcinoma via a positive feedback loop with TGF-β.
Cancer letters
Meng L,Zheng Y,Liu S,Ju Y,Ren S,Sang Y,Zhu Y,Gu L,Liu F,Zhao Y,Zhang X,Sang M
Thu Oct 28 00:00:00 UTC 2021
Hepatic Ischemia-Reperfusion Impairs Blood-Brain Barrier Partly Due to Release of Arginase From Injured Liver.
Frontiers in pharmacology
Zhu L,Zhou H,Xu F,Yang H,Li P,Sheng Y,Liu P,Kong W,Liu X,Yang L,Liu L,Liu X
Tue Nov 02 00:00:00 UTC 2021
Lidocaine Promoted Ferroptosis by Targeting miR-382-5p /SLC7A11 Axis in Ovarian and Breast Cancer.
Frontiers in pharmacology
Sun D,Li YC,Zhang XY
Tue Jun 15 00:00:00 UTC 2021
Probiotic fermentation of Ganoderma lucidum fruiting body extracts promoted its immunostimulatory activity in mice with dexamethasone-induced immunosuppression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li Y,Liu H,Qi H,Tang W,Zhang C,Liu Z,Liu Y,Wei X,Kong Z,Jia S,Du B,Yuan J,Wang C,Li M
Wed Sep 01 00:00:00 UTC 2021
Saikosaponin D Inhibits the Proliferation and Promotes the Apoptosis of Rat Hepatic Stellate Cells by Inducing Autophagosome Formation.
Evidence-based complementary and alternative medicine : eCAM
Jiang H,Liu J,Zhang K,Zeng Q
Tue Apr 26 00:00:00 UTC 2022
Ferroptosis is involved in the anti-tumor effect of lycorine in renal cell carcinoma cells.
Oncology letters
Du Y,Zhao HC,Zhu HC,Jin Y,Wang L
Mon Nov 01 00:00:00 UTC 2021
Chrysanthemi Flos extract alleviated acetaminophen-induced rat liver injury via inhibiting oxidative stress and apoptosis based on network pharmacology analysis.
Pharmaceutical biology
Zhou Y,Wang C,Kou J,Wang M,Rong X,Pu X,Xie X,Han G,Pang X
Wed Dec 01 00:00:00 UTC 2021
Growth hormone receptor disrupts glucose homeostasis via promoting and stabilizing retinol binding protein 4.
Theranostics
Liu J,Nie C,Xue L,Yan Y,Liu S,Sun J,Fan M,Qian H,Ying H,Wang L,Li Y
Wed Jan 19 00:00:00 UTC 2022
Low expression of BTN3A3 indicates poor prognosis and promotes cell proliferation, migration and invasion in non-small cell lung cancer.
Annals of translational medicine
Cheng X,Ma T,Yi L,Su C,Wang X,Wen T,Wang B,Wang Y,Zhang H,Liu Z
Mon Mar 01 00:00:00 UTC 2021
MicroRNA-30c-5p protects against myocardial ischemia/reperfusion injury via regulation of Bach1/Nrf2.
Toxicology and applied pharmacology
Sun M,Guo M,Ma G,Zhang N,Pan F,Fan X,Wang R
Wed Sep 01 00:00:00 UTC 2021
STAT5 inhibitor attenuates atherosclerosis via inhibition of inflammation: the role of STAT5 in atherosclerosis.
American journal of translational research
Wang X,Ding X,Yan J,Lu Z,Cao H,Ni X,Ying Y
Tue Apr 13 00:00:00 UTC 2021
Reactive microglia enhance the transmission of exosomal α-synuclein via toll-like receptor 2.
Brain : a journal of neurology
Xia Y,Zhang G,Kou L,Yin S,Han C,Hu J,Wan F,Sun Y,Wu J,Li Y,Huang J,Xiong N,Zhang Z,Wang T
Tue Aug 17 00:00:00 UTC 2021
Diosmetin ameliorate type 2 diabetic mellitus by up-regulating Corynebacterium glutamicum to regulate IRS/PI3K/AKT-mediated glucose metabolism disorder in KK-Ay mice.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Gong X,Xiong L,Bi C,Zhang B
Thu Jul 01 00:00:00 UTC 2021
Zinc-α2-glycoprotein relieved seizure-Induced neuronal glucose uptake impairment via insulin-like growth factor 1 receptor-regulated glucose transporter 3 expression.
Journal of neurochemistry
Peng W,Liu X,Tan C,Zhou W,Jiang J,Zhou X,Du J,Mo L,Chen L
Sat May 01 00:00:00 UTC 2021
Contribution of the NLRP3/IL-1β axis to impaired vasodilation in sepsis through facilitation of eNOS proteolysis and the protective role of melatonin.
International immunopharmacology
Hu S,Pi Q,Luo M,Cheng Z,Liang X,Luo S,Xia Y
Thu Apr 01 00:00:00 UTC 2021
Impact of knockdown LincRNA-Cox2 on apoptosis of macrophage infected with Bacillus Calmette-Guérin.
Molecular immunology
Xu Y,Yu J,Ma C,Gong Z,Wu X,Deng G
Mon Feb 01 00:00:00 UTC 2021
Epigenetically silenced linc00261 contributes to the metastasis of hepatocellular carcinoma via inducing the deficiency of FOXA2 transcription.
American journal of cancer research
Chen Z,Xiang L,Hu Z,Ou H,Liu X,Yu L,Chen W,Jiang L,Yu Q,Fang Y,Xu Y,Liu Q,Huang Y,Li X,Yang D
Tue Feb 02 00:00:00 UTC 2021
A clinically acceptable strategy for sensitizing anti-PD-1 treatment by hypoxia relief.
Journal of controlled release : official journal of the Controlled Release Society
Jiang M,Qin B,Luo L,Li X,Shi Y,Zhang J,Luo Z,Zhu C,Guan G,Du Y,You J
Sat Jul 10 00:00:00 UTC 2021
PET-CT and RNA sequencing reveal novel targets for acupuncture-induced lowering of blood pressure in spontaneously hypertensive rats.
Scientific reports
Li J,Peng C,Lai D,Fang Y,Luo D,Zhou Z,Li C,Lai X
Wed May 26 00:00:00 UTC 2021
CircCRIM1 promotes ovarian cancer progression by working as ceRNAs of CRIM1 and targeting miR-383-5p/ZEB2 axis.
Reproductive biology and endocrinology : RB&E
Du Y,Liu X,Zhang S,Chen S,Guan X,Li Q,Chen X,Zhao Y
Tue Nov 30 00:00:00 UTC 2021
Autocrined leptin promotes proliferation of non-small cell lung cancer (NSCLC) via PI3K/AKT and p53 pathways.
Annals of translational medicine
Wang J,Zhou F,Li F,Wang B,Hu Y,Li X
Thu Apr 01 00:00:00 UTC 2021
Identification of a Proteomic Signature of Senescence in Primary Human Mammary Epithelial Cells.
Journal of proteome research
Delfarah A,Hartel NG,Zheng D,Yang J,Graham NA
Fri Nov 05 00:00:00 UTC 2021
The Formation of Melanocyte Apoptotic Bodies in Vitiligo and the Relocation of Vitiligo Autoantigens under Oxidative Stress.
Oxidative medicine and cellular longevity
Tian J,Wang Y,Ding M,Zhang Y,Chi J,Wang T,Jiao B,Jian Z,Yi X,Huang Y,Liu L,Li K,Chen J,Wang G,Gao T,Li C,Li Q
Thu Feb 17 00:00:00 UTC 2022
Ramelteon Mitigates Free Fatty Acid (FFA)-Induced Attachment of Monocytes to Brain Vascular Endothelial Cells.
Neurotoxicity research
Wang G,Tian F,Li Y,Liu Y,Liu C
Wed Dec 01 00:00:00 UTC 2021
Phosphatidylinositol 3,4,5-Trisphosphate-Dependent Rac Exchanger 2 Protein Facilitates Glioma Progression via Akt and Stat3 Signaling.
Journal of molecular neuroscience : MN
Shang J,Li Y,Yin G,Li Z,Jiang L,Bai Q
Sun Aug 01 00:00:00 UTC 2021
Downregulated fat mass and obesity-associated protein inhibits bone resorption and osteoclastogenesis by nuclear factor-kappa B inactivation.
Cellular signalling
Zhuang J,Ning H,Wang M,Zhao W,Jing Y,Liu X,Zu J,Kong P,Wang X,Sun C,Yan J
Mon Nov 01 00:00:00 UTC 2021
A novel peptide relieves endothelial cell dysfunction in preeclampsia by regulating the PI3K/mTOR/HIF1α pathway.
International journal of molecular medicine
Xue L,Xie K,Wu L,Yu X,Long W,Li C,Jia R,Ding H
Fri Jan 01 00:00:00 UTC 2021
Zinc provides neuroprotection by regulating NLRP3 inflammasome through autophagy and ubiquitination in a spinal contusion injury model.
CNS neuroscience & therapeutics
Lin JQ,Tian H,Zhao XG,Lin S,Li DY,Liu YY,Xu C,Mei XF
Thu Apr 01 00:00:00 UTC 2021
The Abscopal Effects of Cranial Irradiation Induce Testicular Damage in Mice.
Frontiers in physiology
Guo L,Qin TZ,Liu LY,Lai PP,Xue YZ,Jing YT,Zhang W,Li W,Li J,Ding GR
Tue Dec 07 00:00:00 UTC 2021
Spleen Tyrosine Kinase Mediates Microglial Activation in Mice With Diabetic Retinopathy.
Translational vision science & technology
Liu X,Xu B,Gao S
Thu Apr 01 00:00:00 UTC 2021
Antioral Squamous Cell Carcinoma Effects of Carvacrol via Inhibiting Inflammation, Proliferation, and Migration Related to Nrf2/Keap1 Pathway.
BioMed research international
Liu H,Xu X,Wu R,Bi L,Zhang C,Chen H,Yang Y
Mon Oct 25 00:00:00 UTC 2021
Effect of claudin 1 on cell proliferation, migration and apoptosis in human cervical squamous cell carcinoma.
Oncology reports
Shen Z,Song W,Qian L,Zhu J,Li Y,Li M,Zhang T,Zhao W,Zhou Y,Yang X
Mon Feb 01 00:00:00 UTC 2021
Angiotensin II-mediated improvement of renal mitochondrial function via the AMPK/PGC-1α/NRF-2 pathway is superior to norepinephrine in a rat model of septic shock associated with acute renal injury.
Annals of translational medicine
An H,Hu Z,Chen Y,Cheng L,Shi J,Han L
Mon Mar 01 00:00:00 UTC 2021
Acetabular development and fate of inverted limbus in rabbits: Experimental observation from an animal model.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Liu J,Zhou W,Chen Y,Li L
Wed Dec 01 00:00:00 UTC 2021
miR-320a in serum exosomes promotes myocardial fibroblast proliferation via regulating the PIK3CA/Akt/mTOR signaling pathway in HEH2 cells.
Experimental and therapeutic medicine
Wang QG,Cheng BC,He YZ,Li LJ,Ling Y,Luo G,Wang L,Liang S,Zhang Y
Sun Aug 01 00:00:00 UTC 2021
Celastrol Inhibits Canine Mammary Tumor Cells by Inducing Apoptosis via the Caspase Pathway.
Frontiers in veterinary science
Ou G,Jiang X,Gao A,Li X,Lin Z,Pei S
Wed Feb 23 00:00:00 UTC 2022
SPOP negatively regulates Toll-like receptor-induced inflammation by disrupting MyD88 self-association.
Cellular & molecular immunology
Hu YH,Wang Y,Wang F,Dong YM,Jiang WL,Wang YP,Zhong X,Ma LX
Thu Jul 01 00:00:00 UTC 2021
IKKβ activation promotes amphisome formation and extracellular vesicle secretion in tumor cells.
Biochimica et biophysica acta. Molecular cell research
Peng X,Yang L,Ma Y,Li X,Yang S,Li Y,Wu B,Tang S,Zhang F,Zhang B,Liu J,Li H
Fri Jan 01 00:00:00 UTC 2021
TRIP4 transcriptionally activates DDIT4 and subsequent mTOR signaling to promote glioma progression.
Free radical biology & medicine
Li W,Hu S,Tian C,Wan X,Yu W,Guo P,Zhao F,Hua C,Lu X,Xue G,Han S,Guo W,Wang D,Deng W
Wed Dec 01 00:00:00 UTC 2021
Dental pulp stem cell-derived exosomes alleviate cerebral ischaemia-reperfusion injury through suppressing inflammatory response.
Cell proliferation
Li S,Luo L,He Y,Li R,Xiang Y,Xing Z,Li Y,Albashari AA,Liao X,Zhang K,Gao L,Ye Q
Sun Aug 01 00:00:00 UTC 2021
A Mixture of Baicalein, Wogonin, and Oroxylin-A Inhibits EMT in the A549 Cell Line via the PI3K/AKT-TWIST1-Glycolysis Pathway.
Frontiers in pharmacology
Cao HJ,Zhou W,Xian XL,Sun SJ,Ding PJ,Tian CY,Tian FL,Jiang CH,Fu TT,Zhao S,Dai JY
Tue Mar 01 00:00:00 UTC 2022
KIM-1-mediated anti-inflammatory activity is preserved by MUC1 induction in the proximal tubule during ischemia-reperfusion injury.
American journal of physiology. Renal physiology
Al-Bataineh MM,Kinlough CL,Mi Z,Jackson EK,Mutchler SM,Emlet DR,Kellum JA,Hughey RP
Sun Aug 01 00:00:00 UTC 2021
GDF11 prevents the formation of thoracic aortic dissection in mice: Promotion of contractile transition of aortic SMCs.
Journal of cellular and molecular medicine
Ren K,Li B,Liu Z,Xia L,Zhai M,Wei X,Duan W,Yu S
Sat May 01 00:00:00 UTC 2021
Morusin inhibits the growth of human colorectal cancer HCT116‑derived sphere‑forming cells via the inactivation of Akt pathway.
International journal of molecular medicine
Zhou Y,Li X,Ye M
Thu Apr 01 00:00:00 UTC 2021
Integrating systematic pharmacology-based strategy and experimental validation to explore the synergistic pharmacological mechanisms of Guanxin V in treating ventricular remodeling.
Bioorganic chemistry
Liang B,Liang Y,Li R,Zhang H,Gu N
Fri Oct 01 00:00:00 UTC 2021
Syndecan 4 contributes to osteoclast differentiation induced by RANKL through enhancing autophagy.
International immunopharmacology
Li J,Sun Z,Lin Y,Yan Y,Yan H,Jing B,Han Z
Mon Feb 01 00:00:00 UTC 2021
Biomimetic amphiphilic FAAP NPs nanoparticles: Synthesis, characterization and antivirus activity.
International immunopharmacology
Li XH,Zhang Z,Ma X,Guo ZH,Ban X,Tian Z,Zhao L,Liu Q,He Q
Wed Dec 01 00:00:00 UTC 2021
GR/NF-κB signaling pathway regulates hippocampal inflammatory responses in diabetic rats with chronic unpredictable mild stress.
European journal of pharmacology
Li ZR,Han YS,Liu Z,Zhao HQ,Liu J,Yang H,Wang YH
Mon Mar 15 00:00:00 UTC 2021
Transcriptome Differences in Pig Tracheal Epithelial Cells in Response to Pasteurella Multocida Infection.
Frontiers in veterinary science
Wang X,Wang F,Lin L,Liang W,Liu S,Hua L,Wang X,Chen H,Peng Z,Wu B
Wed Sep 08 00:00:00 UTC 2021
Xanthine oxidase inhibitory activity and antihyperuricemic effect of Moringa oleifera Lam. leaf hydrolysate rich in phenolics and peptides.
Journal of ethnopharmacology
Tian Y,Lin L,Zhao M,Peng A,Zhao K
Sat Apr 24 00:00:00 UTC 2021
Sesamin Protects against and Ameliorates Rat Intestinal Ischemia/Reperfusion Injury with Involvement of Activating Nrf2/HO-1/NQO1 Signaling Pathway.
Oxidative medicine and cellular longevity
Wang Y,Wen J,Almoiliqy M,Wang Y,Liu Z,Yang X,Lu X,Meng Q,Peng J,Lin Y,Sun P
Fri Feb 04 00:00:00 UTC 2022
LncRNA-AC006129.1 reactivates a SOCS3-mediated anti-inflammatory response through DNA methylation-mediated CIC downregulation in schizophrenia.
Molecular psychiatry
Ni C,Jiang W,Wang Z,Wang Z,Zhang J,Zheng X,Liu Z,Ou H,Jiang T,Liang W,Wu F,Li Q,Hou Y,Yang Q,Guo B,Liu S,Li S,Li S,Yang E,Zhu XH,Huang X,Wen Z,Zhao C
Sun Aug 01 00:00:00 UTC 2021
The Performance of HepG2 and HepaRG Systems through the Glass of Acetaminophen-Induced Toxicity.
Life (Basel, Switzerland)
Lőrincz T,Deák V,Makk-Merczel K,Varga D,Hajdinák P,Szarka A
Sat Aug 21 00:00:00 UTC 2021
Curcumol inhibits ferritinophagy to restrain hepatocyte senescence through YAP/NCOA4 in non-alcoholic fatty liver disease.
Cell proliferation
Qi X,Song A,Ma M,Wang P,Zhang X,Lu C,Zhang J,Zheng S,Jin H
Wed Sep 01 00:00:00 UTC 2021
Human umbilical cord mesenchymal stem cells ameliorate depression by regulating Jmjd3 and microglia polarization in myocardial infarction mice.
Psychopharmacology
Zhang Y,Wang X,Li Y,Liu R,Pan J,Tang X,Sun S,Liu J,Ma W
Fri Oct 01 00:00:00 UTC 2021
PHLDA3 inhibition protects against myocardial ischemia/reperfusion injury by alleviating oxidative stress and inflammatory response via the Akt/Nrf2 axis.
Environmental toxicology
Meng X,Zhang L,Han B,Zhang Z
Mon Nov 01 00:00:00 UTC 2021
Apigenin Alleviates Intervertebral Disc Degeneration via Restoring Autophagy Flux in Nucleus Pulposus Cells.
Frontiers in cell and developmental biology
Xie C,Shi Y,Chen Z,Zhou X,Luo P,Hong C,Tian N,Wu Y,Zhou Y,Lin Y,Dou H,Wu A,Huang Q,Zhang X,Wang X
Tue Feb 01 00:00:00 UTC 2022
LncRNA WEE2-AS1 knockdown inhibits the proliferation, migration and invasion of glioma cells via regulating miR-29b-2-5p/TPM3 axis.
Oncology research
Jia Z,Qian Z,Tang Y,Li X,Shi Y,Xin H,Fan Y,Wu H
Tue Jun 13 00:00:00 UTC 2023
Exposure to melamine cyanuric acid in adult mice induced thyroid dysfunction and circadian rhythm disorder.
Ecotoxicology and environmental safety
Wang S,Wang S,Wang C,Feng D,Feng X
Fri Nov 19 00:00:00 UTC 2021
Luteolin attenuates angiotensin II‑induced renal damage in apolipoprotein E‑deficient mice.
Molecular medicine reports
Liu YS,Yang Q,Li S,Luo L,Liu HY,Li XY,Gao ZN
Mon Feb 01 00:00:00 UTC 2021
SIRT1 Attenuates Apoptosis of Nucleus Pulposus Cells by Targeting Interactions between LC3B and Fas under High-Magnitude Compression.
Oxidative medicine and cellular longevity
Zhuo Y,Wang H,Zou L,Wang Y,Hu Y,Li P,Zhou Q
Fri Mar 04 00:00:00 UTC 2022
CENPE contributes to pulmonary vascular remodeling in pulmonary hypertension.
Biochemical and biophysical research communications
Fang X,Xie M,Liu X,He Y
Fri Jun 11 00:00:00 UTC 2021
Protective Effects of Huangqi Shengmai Yin on Type 1 Diabetes-Induced Cardiomyopathy by Improving Myocardial Lipid Metabolism.
Evidence-based complementary and alternative medicine : eCAM
Cao Z,Pan J,Sui X,Fang C,Li N,Huang X,Qu X,Han D
Tue Jul 13 00:00:00 UTC 2021
Histone demethylase JMJD3 protects against renal fibrosis by suppressing TGFβ and Notch signaling and preserving PTEN expression.
Theranostics
Yu C,Xiong C,Tang J,Hou X,Liu N,Bayliss G,Zhuang S
Tue Jul 27 00:00:00 UTC 2021
Magnesium-L-threonate alleviate colonic inflammation and memory impairment in chronic-plus-binge alcohol feeding mice.
Brain research bulletin
Liu C,Cheng Y,Guo Y,Qian H
Wed Sep 01 00:00:00 UTC 2021
Betulinic acid induces autophagy-dependent apoptosis via Bmi-1/ROS/AMPK-mTOR-ULK1 axis in human bladder cancer cells.
Aging
Zhang Y,He N,Zhou X,Wang F,Cai H,Huang SH,Chen X,Hu Z,Jin X
Sun Sep 12 00:00:00 UTC 2021
Cdk5-Mediated Phosphorylation of Sirt1 Contributes to Podocyte Mitochondrial Dysfunction in Diabetic Nephropathy.
Antioxidants & redox signaling
Wang S,Yang Y,He X,Yang L,Wang J,Xia S,Liu D,Liu S,Yang L,Liu W,Duan H
Wed Jan 20 00:00:00 UTC 2021
Astragaloside IV inhibits cell proliferation and metastasis of breast cancer via promoting the long noncoding RNA TRHDE-AS1.
Journal of natural medicines
Hu S,Zheng W,Jin L
Fri Jan 01 00:00:00 UTC 2021
Myosin 1f-mediated activation of microglia contributes to the photoreceptor degeneration in a mouse model of retinal detachment.
Cell death & disease
Wang Y,Zhao X,Gao M,Wan X,Guo Y,Qu Y,Chen Y,Li T,Liu H,Jiang M,Wang F,Sun X
Sat Oct 09 00:00:00 UTC 2021
YTHDF1 promotes hepatocellular carcinoma progression via activating PI3K/AKT/mTOR signaling pathway and inducing epithelial-mesenchymal transition.
Experimental hematology & oncology
Luo X,Cao M,Gao F,He X
Fri Jun 04 00:00:00 UTC 2021
HMSC-Derived Exosome Inhibited Th2 Cell Differentiation via Regulating miR-146a-5p/SERPINB2 Pathway.
Journal of immunology research
Zhou J,Lu Y,Wu W,Feng Y
Mon Dec 06 00:00:00 UTC 2021
AP39 ameliorates high fat diet-induced liver injury in young rats via alleviation of oxidative stress and mitochondrial impairment.
Experimental animals
Yu Y,Ye SM,Liu DY,Yang LQ
Wed Nov 10 00:00:00 UTC 2021
Comprehensive Analysis of the Tumor Microenvironment and Ferroptosis-Related Genes Predict Prognosis with Ovarian Cancer.
Frontiers in genetics
Li XX,Xiong L,Wen Y,Zhang ZJ
Tue Dec 07 00:00:00 UTC 2021
GCG inhibits SARS-CoV-2 replication by disrupting the liquid phase condensation of its nucleocapsid protein.
Nature communications
Zhao M,Yu Y,Sun LM,Xing JQ,Li T,Zhu Y,Wang M,Yu Y,Xue W,Xia T,Cai H,Han QY,Yin X,Li WH,Li AL,Cui J,Yuan Z,Zhang R,Zhou T,Zhang XM,Li T
Fri Apr 09 00:00:00 UTC 2021
Bone marrow stem cells therapy alleviates vascular injury in a chronic obstructive pulmonary disease‑obstructive sleep apnea overlap syndrome rat model.
Molecular medicine reports
Bi H,He J,He X,Du J,Chen M,Huang Z,Yang C,Yang L,Li H,Zhou K,Wang Q,He L,Jin Z
Fri Jan 01 00:00:00 UTC 2021
UFL1 regulates milk protein and fat synthesis-related gene expression of bovine mammary epithelial cells probably via the mTOR signaling pathway.
In vitro cellular & developmental biology. Animal
Li C,Li L,Ali I,Kuang M,Wang X,Wang G
Sat May 01 00:00:00 UTC 2021
Recombinant Human Growth Hormone Inhibits Lipotoxicity, Oxidative Stress, and Apoptosis in a Mouse Model of Diabetic Cardiomyopathy.
Oxidative medicine and cellular longevity
Pei Z,Wang X,Yang C,Dong M,Wang F
Mon Feb 28 00:00:00 UTC 2022
A Cas9-transcription factor fusion protein enhances homology-directed repair efficiency.
The Journal of biological chemistry
Li G,Wang H,Zhang X,Wu Z,Yang H
Fri Aug 20 00:00:00 UTC 2021
TDP-43 and PINK1 mediate CHCHD10S59L mutation-induced defects in Drosophila and in vitro.
Nature communications
Baek M,Choe YJ,Bannwarth S,Kim J,Maitra S,Dorn GW,Taylor JP,Paquis-Flucklinger V,Kim NC
Fri Mar 26 00:00:00 UTC 2021
Exosomes derived from idiopathic gingival fibroma fibroblasts regulate gingival fibroblast proliferation and apoptosis.
Oral diseases
Yin S,Jia F,Ran L,Xie L,Wu Z,Zhan Y,Zhang Y,Zhang M
Fri Oct 01 00:00:00 UTC 2021
New targets of morphine postconditioning protection of the myocardium in ischemia/reperfusion injury: Involvement of HSP90/Akt and C5a/NF-κB.
Open medicine (Warsaw, Poland)
Tu RH,Wang DX,Zhong GQ,Meng JJ,Wen H,Bi Q,He Y
Wed Apr 27 00:00:00 UTC 2022
Single-Cell Analysis Identify Transcription Factor BACH1 as a Master Regulator Gene in Vascular Cells During Aging.
Frontiers in cell and developmental biology
Ge F,Pan Q,Qin Y,Jia M,Ruan C,Wei X,Jing Q,Zhi X,Wang X,Jiang L,Osto E,Guo J,Meng D
Tue Jan 11 00:00:00 UTC 2022
Virtual screening identified natural Keap1-Nrf2 PPI inhibitor alleviates inflammatory osteoporosis through Nrf2-mir214-Traf3 axis.
Free radical biology & medicine
Hong J,Shi Z,Li C,Ji X,Li S,Chen Y,Jiang G,Shi M,Wang W,Zhang Y,Hu B,Yan S
Sun Aug 01 00:00:00 UTC 2021
Activation of CB2 receptor inhibits pyroptosis and subsequently ameliorates cecal ligation and puncture-induced sepsis.
International immunopharmacology
Zhang B,Zheng F,Liu A,Li Z,Zheng F,Liu Q,Yang L,Chen K,Wang Y,Zhang Z,He Q,Zhan J
Fri Oct 01 00:00:00 UTC 2021
The p53/p21/p16 and PI3K/Akt signaling pathways are involved in the ameliorative effects of maltol on D-galactose-induced liver and kidney aging and injury.
Phytotherapy research : PTR
Sha JY,Li JH,Zhou YD,Yang JY,Liu W,Jiang S,Wang YP,Zhang R,Di P,Li W
Sun Aug 01 00:00:00 UTC 2021
Tumor protein D52 is upregulated in oral squamous carcinoma cells under hypoxia in a hypoxia-inducible-factor-independent manner and is involved in cell death resistance.
Cell & bioscience
Abe Y,Mukudai Y,Kurihara M,Houri A,Chikuda J,Yaso A,Kato K,Shimane T,Shirota T
Sat Jul 03 00:00:00 UTC 2021
Bach2 overexpression represses Th9 cell differentiation by suppressing IRF4 expression in systemic lupus erythematosus.
FEBS open bio
Sheng Y,Zhang J,Li K,Wang H,Wang W,Wen L,Gao J,Tang X,Tang H,Huang H,Cai M,Yuan T,Liu L,Zheng X,Zhu Z,Cui Y
Mon Feb 01 00:00:00 UTC 2021
KLF5-mediated Eppk1 expression promotes cell proliferation in cervical cancer via the p38 signaling pathway.
BMC cancer
Ma D,Pan Z,Chang Q,Zhang JJ,Liu X,Hua N,Li GH
Thu Apr 08 00:00:00 UTC 2021
Th17/IL-17 induces endothelial cell senescence via activation of NF-κB/p53/Rb signaling pathway.
Laboratory investigation; a journal of technical methods and pathology
Zhang L,Liu M,Liu W,Hu C,Li H,Deng J,Cao Q,Wang Y,Hu W,Li Q
Mon Nov 01 00:00:00 UTC 2021
ACSS3 represses prostate cancer progression through downregulating lipid droplet-associated protein PLIN3.
Theranostics
Zhou L,Song Z,Hu J,Liu L,Hou Y,Zhang X,Yang X,Chen K
Wed Aug 04 00:00:00 UTC 2021
F-Box protein 4 inhibits progression of papillary thyroid cancer.
Steroids
Xu M,Wang Y,Chen M,Hu H,Xia T,Deng D
Mon Feb 01 00:00:00 UTC 2021
Starvation induced autophagy promotes the progression of bladder cancer by LDHA mediated metabolic reprogramming.
Cancer cell international
Li T,Tong H,Yin H,Luo Y,Zhu J,Qin Z,Yin S,He W
Sun Nov 07 00:00:00 UTC 2021
Shenmai injection improves doxorubicin cardiotoxicity via miR-30a/Beclin 1.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang X,Lv S,Zhang W,Jia Q,Wang L,Ding Y,Yuan P,Zhu Y,Liu L,Li Y,Zhang J
Thu Jul 01 00:00:00 UTC 2021
Protein kinase D participates in cardiomyocyte hypertrophy by regulating extracellular signal-regulated and myocyte enhancer factor 2D.
Revista portuguesa de cardiologia
Yuan H,Xiang Q,Yang L,Geng J
Mon Mar 01 00:00:00 UTC 2021
Simiao Qingwen Baidu decoction inhibits Epstein-Barr virus-induced B lymphoproliferative disease and lytic viral replication.
Pharmaceutical biology
Yang X,Liu L,Zhang H,Sun X,Yan Y,Ran R
Wed Dec 01 00:00:00 UTC 2021
Effect of different exercise training intensities on age-related cardiac damage in male mice.
Aging
Pei Z,Yang C,Guo Y,Dong M,Wang F
Tue Sep 14 00:00:00 UTC 2021
Antiviral effects of ergosterol peroxide in a pig model of porcine deltacoronavirus (PDCoV) infection involves modulation of apoptosis and tight junction in the small intestine.
Veterinary research
Duan C,Wang J,Liu Y,Zhang J,Si J,Hao Z,Wang J
Mon Jun 14 00:00:00 UTC 2021
Diosgenin Protects Against Kidney Injury and Mitochondrial Apoptosis Induced by 3-MCPD Through the Regulation of ER Stress, Ca2+ Homeostasis, and Bcl2 Expression.
Molecular nutrition & food research
Zhong Y,Jin C,Han J,Zhu J,Liu Q,Sun D,Xia X,Zhang Y,Peng X
Sun Aug 01 00:00:00 UTC 2021
Posterior Neocortex-Specific Regulation of Neuronal Migration by CEP85L Identifies Maternal Centriole-Dependent Activation of CDK5.
Neuron
Kodani A,Kenny C,Lai A,Gonzalez DM,Stronge E,Sejourne GM,Isacco L,Partlow JN,O'Donnell A,McWalter K,Byrne AB,Barkovich AJ,Yang E,Hill RS,Gawlinski P,Wiszniewski W,Cohen JS,Fatemi SA,Baranano KW,Sahin M,Vossler DG,Yuskaitis CJ,Walsh CA
Wed Apr 22 00:00:00 UTC 2020
Myeloid cells protect intestinal epithelial barrier integrity through the angiogenin/plexin-B2 axis.
The EMBO journal
Bai R,Sun D,Chen M,Shi X,Luo L,Yao Z,Liu Y,Ge X,Gao X,Hu GF,Zhou W,Sheng J,Xu Z
Wed Jul 01 00:00:00 UTC 2020
Saikosaponin-d impedes hippocampal neurogenesis and causes cognitive deficits by inhibiting the survival of neural stem/progenitor cells via neurotrophin receptor signaling in mice.
Clinical and translational medicine
Qin T,Yuan Z,Yu J,Fu X,Deng X,Fu Q,Ma Z,Ma S
Tue Dec 01 00:00:00 UTC 2020
Myeloid-derived suppressor cells promote epithelial ovarian cancer cell stemness by inducing the CSF2/p-STAT3 signalling pathway.
The FEBS journal
Li X,Wang J,Wu W,Gao H,Liu N,Zhan G,Li L,Han L,Guo X
Tue Dec 01 00:00:00 UTC 2020
Inhibition of PFKFB3 suppresses osteoclastogenesis and prevents ovariectomy-induced bone loss.
Journal of cellular and molecular medicine
Wang J,Guan H,Liu H,Lei Z,Kang H,Guo Q,Dong Y,Liu H,Sun Y,Fang Z,Li F
Sat Feb 01 00:00:00 UTC 2020
Deletion of the Impg2 gene causes the degeneration of rod and cone cells in mice.
Human molecular genetics
Xu H,Qu C,Gan L,Sun K,Tan J,Liu X,Jiang Z,Tian W,Liu W,Zhang S,Yang Y,Jiang L,Zhu X,Zhang L
Sat Jun 27 00:00:00 UTC 2020
Fine-tuning of the respiratory complexes stability and supercomplexes assembly in cells defective of complex III.
Biochimica et biophysica acta. Bioenergetics
Tropeano CV,Aleo SJ,Zanna C,Roberti M,Scandiffio L,Loguercio Polosa P,Fiori J,Porru E,Roda A,Carelli V,Steimle S,Daldal F,Rugolo M,Ghelli A
Sat Feb 01 00:00:00 UTC 2020
miR-876 Inhibits EMT and Liver Fibrosis via POSTN to Suppress Metastasis in Hepatocellular Carcinoma.
BioMed research international
Chen K,Li Z,Zhang M,Wang B,Peng T,Shen Y,Zhang J,Ye J,Liu Y,Tang D,Peng M,Ma D,Xiao Z,Zhang Y,Jin W,Li X
Wed May 05 00:00:00 UTC 2021
miR‑95 promotes osteosarcoma growth by targeting SCNN1A.
Oncology reports
Geng Y,Zhao S,Jia Y,Xia G,Li H,Fang Z,Zhang Q,Tian R
Fri May 01 00:00:00 UTC 2020
Sulforaphane enhances the cisplatin sensitivity through regulating DNA repair and accumulation of intracellular cisplatin in ovarian cancer cells.
Experimental cell research
Gong TT,Liu XD,Zhan ZP,Wu QJ
Sat Aug 15 00:00:00 UTC 2020
Matrine suppresses breast cancer metastasis by targeting ITGB1 and inhibiting epithelial-to-mesenchymal transition.
Experimental and therapeutic medicine
Ren L,Mo W,Wang L,Wang X
Wed Jan 01 00:00:00 UTC 2020
Sodium butyrate inhibits migration and induces AMPK-mTOR pathway-dependent autophagy and ROS-mediated apoptosis via the miR-139-5p/Bmi-1 axis in human bladder cancer cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wang F,Wu H,Fan M,Yu R,Zhang Y,Liu J,Zhou X,Cai Y,Huang S,Hu Z,Jin X
Sun Mar 01 00:00:00 UTC 2020
CADM1 inhibits ovarian cancer cell proliferation and migration by potentially regulating the PI3K/Akt/mTOR pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Si X,Xu F,Xu F,Wei M,Ge Y,Chenge S
Sun Mar 01 00:00:00 UTC 2020
Bufalin Induces Glioma Cell Death by Apoptosis or Necroptosis.
OncoTargets and therapy
LingHu HR,Luo H,Gang L
Fri Apr 15 00:00:00 UTC 2022
Naturally-derived diterpenoid sphaeropsidin C as an activator of Nrf2/ARE pathway and its potential capability of relieving intracellular oxidative stress in human lung epithelial cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li GH,Li AL,Chen XM,Zhang LY,Wang T,Wang XN,Shen T
Wed Jan 01 00:00:00 UTC 2020
Triiodothyronine potentiates angiogenesis-related factor expression through PI3K/AKT signaling pathway in human osteoarthritic osteoblasts.
Iranian journal of basic medical sciences
Li L,Pang Y,Zhang L,Li M,Zhu C,Fang S,Yin Z
Mon Jun 01 00:00:00 UTC 2020
Xiao-Qing-Long-Tang Maintains Cardiac Function during Heart Failure with Reduced Ejection Fraction in Salt-Sensitive Rats by Regulating the Imbalance of Cardiac Sympathetic Innervation.
Evidence-based complementary and alternative medicine : eCAM
Li Z,Wang Y,Jiang Y,Ma D,Jiang P,Zhou G,Yang J,Dong F,Zhao H,Zhang Y,Li X
Wed Apr 20 00:00:00 UTC 2022
Structure-based discovery of neoandrographolide as a novel inhibitor of Rab5 to suppress cancer growth.
Computational and structural biotechnology journal
Zhang J,Sun Y,Zhong LY,Yu NN,Ouyang L,Fang RD,Wang Y,He QY
Sat Dec 19 00:00:00 UTC 2020
The Clinical Relevance and Function of Krüppel-Like Factor 16 in Breast Cancer.
Cancer management and research
Bang S,Li J,Zhang M,Cui R,Wu X,Xin Z,Ma D,Zhang J,Zhang H
Sat Apr 16 00:00:00 UTC 2022
Elevated expression of minichromosome maintenance 3 indicates poor outcomes and promotes G1/S cell cycle progression, proliferation, migration and invasion in colorectal cancer.
Bioscience reports
Zhou H,Xiong Y,Zhang G,Liu Z,Li L,Hou S,Zhou T
Fri Jul 31 00:00:00 UTC 2020
CD4+ T cells and TGFβ1/MAPK signal pathway involved in the valvular hyperblastosis and fibrosis in patients with rheumatic heart disease.
Experimental and molecular pathology
Zhao Z,He D,Ling F,Chu T,Huang D,Wu H,Ge J
Mon Jun 01 00:00:00 UTC 2020
LncRNA TROJAN promotes proliferation and resistance to CDK4/6 inhibitor via CDK2 transcriptional activation in ER+ breast cancer.
Molecular cancer
Jin X,Ge LP,Li DQ,Shao ZM,Di GH,Xu XE,Jiang YZ
Mon May 11 00:00:00 UTC 2020
Study of the protective effect on damaged intestinal epithelial cells of rat multilineage-differentiating stress-enduring (Muse) cells.
Cell biology international
Sun D,Yang L,Cao H,Shen ZY,Song HL
Sat Feb 01 00:00:00 UTC 2020
Hyperbaric oxygen treatment mitigates liver damage in mice with noise exposure.
Clinical and experimental pharmacology & physiology
Meng X,Zhang X,Su Y,Gu Z,Xie X,Chang Y,Bao J
Tue Sep 01 00:00:00 UTC 2020
PRDM16 orchestrates angiogenesis via neural differentiation in the developing brain.
Cell death and differentiation
Su L,Lei X,Ma H,Feng C,Jiang J,Jiao J
Sat Aug 01 00:00:00 UTC 2020
Targeting the miR-122/PKM2 autophagy axis relieves arsenic stress.
Journal of hazardous materials
Wang Y,Zhao H,Guo M,Fei D,Zhang L,Xing M
Wed Feb 05 00:00:00 UTC 2020
miRNA-122-5p stimulates the proliferation and DNA synthesis and inhibits the early apoptosis of human spermatogonial stem cells by targeting CBL and competing with lncRNA CASC7.
Aging
Zhou F,Chen W,Cui Y,Liu B,Yuan Q,Li Z,He Z
Tue Nov 24 00:00:00 UTC 2020
FB-15 inhibits MGC-803 cells growth by regulating energy metabolism.
Chemico-biological interactions
Deng X,Pi Y,Li Z,Xiong R,Liu J,Zhao J,Xie Z,Lei X,Tang G
Tue Aug 25 00:00:00 UTC 2020
HOTAIR maintains the stemness of ovarian cancer stem cells via the miR-206/TBX3 axis.
Experimental cell research
Zhang Y,Guo J,Cai E,Cai J,Wen Y,Lu S,Li X,Han Q,Jiang J,Li T,Wang Z
Thu Oct 15 00:00:00 UTC 2020
Porcine parvovirus nonstructural protein NS1 activates NF-κB and it involves TLR2 signaling pathway.
Journal of veterinary science
Jin X,Yuan Y,Zhang C,Zhou Y,Song Y,Wei Z,Zhang G
Fri May 01 00:00:00 UTC 2020
Identification of novel variants in the FZD4 gene associated with familial exudative vitreoretinopathy in Chinese families.
Clinical & experimental ophthalmology
Xu H,Zhang S,Huang L,Zhao P,Zhang X,Yang Z,Zhang L
Wed Apr 01 00:00:00 UTC 2020
Effect of prednisolone on glyoxalase 1 in an inbred mouse model of aristolochic acid nephropathy using a proteomics method with fluorogenic derivatization-liquid chromatography-tandem mass spectrometry.
PloS one
Chen SM,Lin CE,Chen HH,Cheng YF,Cheng HW,Imai K
Thu Apr 16 00:00:00 UTC 2020
Exogenous hydrogen sulfide inhibits human melanoma cell development via suppression of the PI3K/AKT/ mTOR pathway.
Journal of dermatological science
Xiao Q,Ying J,Qiao Z,Yang Y,Dai X,Xu Z,Zhang C,Xiang L
Wed Apr 01 00:00:00 UTC 2020
Transactivation of miR-202-5p by Steroidogenic Factor 1 (SF1) Induces Apoptosis in Goat Granulosa Cells by Targeting TGFβR2.
Cells
Ding Q,Jin M,Wang Y,Liu J,Kalds P,Wang Y,Yang Y,Wang X,Chen Y
Fri Feb 14 00:00:00 UTC 2020
Discovery of potent small molecule PROTACs targeting mutant EGFR.
European journal of medicinal chemistry
Zhao HY,Yang XY,Lei H,Xi XX,Lu SM,Zhang JJ,Xin M,Zhang SQ
Tue Dec 15 00:00:00 UTC 2020
lncRNA MIRF Promotes Cardiac Apoptosis through the miR-26a-Bak1 Axis.
Molecular therapy. Nucleic acids
Su X,Lv L,Li Y,Fang R,Yang R,Li C,Li T,Zhu D,Li X,Zhou Y,Shan H,Liang H
Fri Jun 05 00:00:00 UTC 2020
The Immunomodulatory Effects and Mechanisms of Tim-3 Action in the Early Stage of Mice with Severe Acute Pancreatitis.
Iranian journal of immunology : IJI
Lin M,Huang J,Chen WC,Fan ZN,Qin X
Sun Mar 01 00:00:00 UTC 2020
Effects of dexmedetomidine on oxidative stress and inflammatory response in lungs during mechanical ventilation in COPD rats.
Experimental and therapeutic medicine
Li P,Han J,Zhang D,Cao S,Su C
Sat Feb 01 00:00:00 UTC 2020
Protective effects of isorhamnetin on pulmonary arterial hypertension: in vivo and in vitro studies.
Phytotherapy research : PTR
Chang Z,Wang JL,Jing ZC,Ma P,Xu QB,Na JR,Tian J,Ma X,Zhou W,Zhou R
Thu Oct 01 00:00:00 UTC 2020
Dl-3-n-Butylphthalide Promotes Remyelination and Suppresses Inflammation by Regulating AMPK/SIRT1 and STAT3/NF-κB Signaling in Chronic Cerebral Hypoperfusion.
Frontiers in aging neuroscience
Li M,Meng N,Guo X,Niu X,Zhao Z,Wang W,Xie X,Lv P
Mon Sep 28 00:00:00 UTC 2020
Elevated levels of arachidonic acid metabolites in follicular fluid of PCOS patients.
Reproduction (Cambridge, England)
Li S,Qi J,Tao Y,Zhu Q,Huang R,Liao Y,Yue J,Liu W,Zhao H,Yin H,Sun Y
Sat Feb 01 00:00:00 UTC 2020
FLI1 promotes protein translation via the transcriptional regulation of MKNK1 expression.
International journal of oncology
Wang C,Song J,Liu W,Yao Y,Kapranov P,Sample KM,Gajendran B,Zacksenhaus E,Hao X,Ben-David Y
Sat Feb 01 00:00:00 UTC 2020
The role of PSMB5 in sodium arsenite-induced oxidative stress in L-02 cells.
Cell stress & chaperones
Lv Y,Hu Q,Shi M,Wang W,Zheng Y,Yang Z,Peng L,Bi D,Zhang A,Hu Y
Fri May 01 00:00:00 UTC 2020
Huoxuezhitong capsule ameliorates MIA-induced osteoarthritis of rats through suppressing PI3K/ Akt/ NF-κB pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ju L,Hu P,Chen P,Xue X,Li Z,He F,Qiu Z,Cheng J,Huang F
Tue Sep 01 00:00:00 UTC 2020
MicroRNA-375 Targets ATG14 to Inhibit Autophagy and Sensitize Hepatocellular Carcinoma Cells to Sorafenib.
OncoTargets and therapy
Yang S,Wang M,Yang L,Li Y,Ma Y,Peng X,Li X,Li B,Jin H,Li H
Thu Apr 14 00:00:00 UTC 2022
Inhibition of TBK1 by amlexanox attenuates paraquat-induced acute lung injury.
Toxicology
Wang N,Li Y,Wang X,Ma Z,Wang Y,Zhang C,Yuan Y,Zhao M
Thu Oct 01 00:00:00 UTC 2020
Anti-Inflammatory and Antioxidant Effect of Eucommia ulmoides Polysaccharide in Hepatic Ischemia-Reperfusion Injury by Regulating ROS and the TLR-4-NF-κB Pathway.
BioMed research international
Gao W,Feng Z,Zhang S,Wu B,Geng X,Fan G,Duan Y,Li K,Liu K,Peng C
Mon Mar 22 00:00:00 UTC 2021
Baicalin reversal of DNA hypermethylation-associated Klotho suppression ameliorates renal injury in type 1 diabetic mouse model.
Cell cycle (Georgetown, Tex.)
Zhang XT,Wang G,Ye LF,Pu Y,Li RT,Liang J,Wang L,Lee KKH,Yang X
Tue Dec 01 00:00:00 UTC 2020
Glutamate-Cysteine Ligase Catalytic Subunit Attenuated Hepatitis C Virus-Related Liver Fibrosis and Suppressed Endoplasmic Reticulum Stress.
Frontiers in molecular biosciences
Fu N,Li D,Li W,Zhao W,Zhang S,Liu L,Zhao S,Du J,Kong L,Wang R,Zhang Y,Nan Y
Tue Oct 06 00:00:00 UTC 2020
Multi-walled carbon nanotubes decrease neuronal NO synthase in 3D brain organoids.
The Science of the total environment
Jiang Y,Gong H,Jiang S,She C,Cao Y
Tue Dec 15 00:00:00 UTC 2020
Effects of the linoleic acid/docosahexaenoic acid ratio and concentration inducing autophagy in Raw264.7 cells against Staphylococcus aureus.
Journal of clinical biochemistry and nutrition
Xu LY,Mu M,Wang ML,Liu JC,Zhou YJ,Wu J,Jiang BY,Chen MG,Hu D,Tao XR
Tue Sep 01 00:00:00 UTC 2020
Hypoxia-induced MFAP5 Promotes Tumor Migration and Invasion via AKT Pathway in Head and Neck Squamous Cell Carcinoma.
Journal of Cancer
Xu Q,Chang H,Tian X,Lou C,Ma H,Yang X
Mon Sep 28 00:00:00 UTC 2020
Fluoride induced mitochondrial impairment and PINK1-mediated mitophagy in Leydig cells of mice: In vivo and in vitro studies.
Environmental pollution (Barking, Essex : 1987)
Liang C,Gao Y,He Y,Han Y,Manthari RK,Tikka C,Chen C,Wang J,Zhang J
Wed Jan 01 00:00:00 UTC 2020
Effect of stromal cell-derived factor-1/CXCR4 axis in neural stem cell transplantation for Parkinson's disease.
Neural regeneration research
Xu JT,Qian Y,Wang W,Chen XX,Li Y,Li Y,Yang ZY,Song XB,Lu D,Deng XL
Wed Jan 01 00:00:00 UTC 2020
Angiotensin II-Induced vascular remodeling and hypertension involves cathepsin L/V- MEK/ERK mediated mechanism.
International journal of cardiology
Lu Y,Sun X,Peng L,Jiang W,Li W,Yuan H,Cai J
Wed Jan 01 00:00:00 UTC 2020
Obesity induces preadipocyte CD36 expression promoting inflammation via the disruption of lysosomal calcium homeostasis and lysosome function.
EBioMedicine
Luo X,Li Y,Yang P,Chen Y,Wei L,Yu T,Xia J,Ruan XZ,Zhao L,Chen Y
Mon Jun 01 00:00:00 UTC 2020
Role of PEPT1in the transport of cinnabar in Caco-2 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Wu Q,He X,Zhou S,Shi F,Lu Y
Sun Mar 01 00:00:00 UTC 2020
LINC00365-SCGB2A1 axis inhibits the viability of breast cancer through targeting NF-κB signaling.
Oncology letters
Zhang L,Yan X,Yu S,Zhong X,Tian R,Xu L,Bian X,Su J
Wed Jan 01 00:00:00 UTC 2020
Insulin Promotes Corneal Nerve Repair and Wound Healing in Type 1 Diabetic Mice by Enhancing Wnt/β-Catenin Signaling.
The American journal of pathology
Yang S,Zhang Y,Zhang Z,Dan J,Zhou Q,Wang X,Li W,Zhou L,Yang L,Xie L
Sun Nov 01 00:00:00 UTC 2020
Effect of STAT3 mediated epigenetic regulation in pre-eclampsia: an analysis of trial data.
European review for medical and pharmacological sciences
Zhang XL,Pan J,Wang G,Li H,Wang JL,Jiang XM,Zhao XY
Mon Jun 01 00:00:00 UTC 2020
Paeoniflorin-6'-O-benzene sulfonate down-regulates CXCR4-Gβγ-PI3K/AKT mediated migration in fibroblast-like synoviocytes of rheumatoid arthritis by inhibiting GRK2 translocation.
Biochemical and biophysical research communications
Wang DD,Jiang MY,Wang W,Zhou WJ,Zhang YW,Yang M,Chen JY,Wei W
Thu Jun 04 00:00:00 UTC 2020
Vernonia patula (Dryand.) Merr. and Leucas chinensis (Retz.) R. Brown exert anti-inflammatory activities and relieve oxidative stress via Nrf2 activation.
Journal of ethnopharmacology
Wu PS,Jeng J,Yang JJ,Kao V,Yen JH,Wu MJ
Sun Nov 15 00:00:00 UTC 2020
The effect of autophagy and mitochondrial fission on Harderian gland is greater than apoptosis in male hamsters during different photoperiods.
PloS one
Xu JH,Wang Z,Mou JJ,Zhao XY,Geng XC,Wu M,Xue HL,Chen L,Xu LX
Thu Dec 24 00:00:00 UTC 2020
Minocycline prevents the depressive-like behavior through inhibiting the release of HMGB1 from microglia and neurons.
Brain, behavior, and immunity
Wang B,Huang X,Pan X,Zhang T,Hou C,Su WJ,Liu LL,Li JM,Wang YX
Sat Aug 01 00:00:00 UTC 2020
Heme Oxygenase-1 Suppresses Wnt Signaling Pathway in Nonalcoholic Steatohepatitis-Related Liver Fibrosis.
BioMed research international
Du J,Ren W,Zhang Q,Fu N,Han F,Cui P,Li W,Kong L,Zhao S,Wang R,Zhang Y,Yang L,Kong L,Nan Y
Fri Mar 12 00:00:00 UTC 2021
Initiation of PI3K/AKT pathway by IGF-1 decreases spinal cord injury-induced endothelial apoptosis and microvascular damage.
Life sciences
Li H,Kong R,Wan B,Yang L,Zhang S,Cao X,Chen H
Tue Dec 15 00:00:00 UTC 2020
The Human-Specific STING Agonist G10 Activates Type I Interferon and the NLRP3 Inflammasome in Porcine Cells.
Frontiers in immunology
Ming SL,Zeng L,Guo YK,Zhang S,Li GL,Ma YX,Zhai YY,Chang WR,Yang L,Wang J,Yang GY,Chu BB
Mon Jun 14 00:00:00 UTC 2021
MicroRNA-128-1-5p attenuates myocardial ischemia/reperfusion injury by suppressing Gadd45g-mediated apoptotic signaling.
Biochemical and biophysical research communications
Wan X,Yao B,Ma Y,Liu Y,Tang Y,Hu J,Li M,Fu S,Zheng X,Yin D
Thu Sep 10 00:00:00 UTC 2020
Critical role of toll-like receptor 4 (TLR4) in ricin toxin-induced inflammatory responses in macrophages.
Toxicology letters
Dong M,Yu H,Wang Y,Sun C,Chang Y,Yin Q,Zhao G,Xu N,Liu W
Sun Mar 15 00:00:00 UTC 2020
Monocyte-derived extracellular vesicles upon treated by palmitate promote endothelial migration and monocytes attachment to endothelial cells.
Biochemical and biophysical research communications
Gao W,Guo X,Wang Y,Jian D,Li M
Thu Mar 12 00:00:00 UTC 2020
AMPK-mTOR-ULK1 axis activation-dependent autophagy promotes hydroxycamptothecin-induced apoptosis in human bladder cancer cells.
Journal of cellular physiology
Wang F,Cao M,Fan M,Wu H,Huang W,Zhang Y,Hu Z,Jin X
Fri May 01 00:00:00 UTC 2020
Norcantharidin protects against renal interstitial fibrosis by suppressing TWEAK-mediated Smad3 phosphorylation.
Life sciences
Zeng D,Xiao Z,Xu Q,Luo H,Wen L,Tang C,Shan Y,Tian J,Wei J,Li Y
Sun Nov 01 00:00:00 UTC 2020
Co-delivery of paclitaxel and anti-VEGF siRNA by tripeptide lipid nanoparticle to enhance the anti-tumor activity for lung cancer therapy.
Drug delivery
Zhang C,Zhao Y,Zhang E,Jiang M,Zhi D,Chen H,Cui S,Zhen Y,Cui J,Zhang S
Tue Dec 01 00:00:00 UTC 2020
Role of mitochondrial acyl-CoA dehydrogenases in the metabolism of dicarboxylic fatty acids.
Biochemical and biophysical research communications
Bharathi SS,Zhang Y,Gong Z,Muzumdar R,Goetzman ES
Thu Jun 18 00:00:00 UTC 2020
HMGB1 mediates homocysteine-induced endothelial cells pyroptosis via cathepsin V-dependent pathway.
Biochemical and biophysical research communications
Leng Y,Chen R,Chen R,He S,Shi X,Zhou X,Zhang Z,Chen AF
Thu Nov 19 00:00:00 UTC 2020
Asperosaponin VI Injection Enhances Orthodontic Tooth Movement in Rats.
Medical science monitor : international medical journal of experimental and clinical research
Ma D,Wang X,Ren X,Bu J,Zheng D,Zhang J
Thu Apr 23 00:00:00 UTC 2020
LMCD1 antisense RNA 1 (LMCD1-AS1) potentiates thyroid cancer cell growth and stemness via a positive feedback loop of LMCD1-AS1/miR-1287-5p/GLI2.
Annals of translational medicine
Shao J,Xu Y,Li H,Chen L,Wang W,Shen D,Chen J
Sun Nov 01 00:00:00 UTC 2020
STAT3-PTTG11 abrogation inhibits proliferation and induces apoptosis in malignant glioma cells.
Oncology letters
Cui L,Xu L,Wang G,Wen J,Luo L,Zhao H,Chen S,Zheng M,Sun C,Jin X,Yang L
Thu Oct 01 00:00:00 UTC 2020
Acylglycerol kinase promotes the stemness of nasopharyngeal carcinoma cells by promoting β-catenin translocation to the nucleus through activating PI3K/Akt pathway.
Environmental toxicology
Zhao Q,Sun P,Qin S,Liu J
Tue Dec 01 00:00:00 UTC 2020
GPAT3 deficiency alleviates insulin resistance and hepatic steatosis in a mouse model of severe congenital generalized lipodystrophy.
Human molecular genetics
Gao M,Liu L,Wang X,Mak HY,Liu G,Yang H
Sat Feb 01 00:00:00 UTC 2020
Expression and distribution of bone morphogenetic protein 4 and its antagonist Noggin in the skin of Kazakh sheep (Ovis aries) with a white and brown coat color.
Acta histochemica
Sun X,Qu G,Wang D,Wang T,Sai W,Chen Y,Yuan L,Pang Q
Fri May 01 00:00:00 UTC 2020
A mutation found in esophageal cancer alters integrin β4 mRNA splicing.
Biochemical and biophysical research communications
Kelly GT,Faraj R,Dai Z,Cress AE,Wang T
Thu Aug 27 00:00:00 UTC 2020
An inducible intestinal epithelial cell-specific NHE3 knockout mouse model mimicking congenital sodium diarrhea.
Clinical science (London, England : 1979)
Xue J,Thomas L,Tahmasbi M,Valdez A,Dominguez Rieg JA,Fenton RA,Rieg T
Thu Apr 30 00:00:00 UTC 2020
Treatment with 5-methoxytryptophan attenuates microglia-induced neuroinflammation in spinal cord trauma.
International immunopharmacology
Hong X,Jiang F,Li Y,Fang L,Qian Z,Chen H,Kong R
Sun Nov 01 00:00:00 UTC 2020
Relevance research between the expression of p16INK4a , Notch1, and hTERC genes: The development of HPV16-positive cervical cancer.
Journal of clinical laboratory analysis
Huo W,Zhai S,Wang Y,Qiang X,Na R,Gui H,Wu N,Cao Y,Bai H
Fri May 01 00:00:00 UTC 2020
Photoperiod Affects Harderian Gland Morphology and Secretion in Female Cricetulus barabensis: Autophagy, Apoptosis, and Mitochondria.
Frontiers in physiology
Wang Z,Xu JH,Mou JJ,Kong XT,Wu M,Xue HL,Xu LX
Mon Sep 28 00:00:00 UTC 2020
Suppression of Notch Signaling Stimulates Progesterone Synthesis by Enhancing the Expression of NR5A2 and NR2F2 in Porcine Granulosa Cells.
Genes
Guo R,Chen F,Shi Z
Wed Jan 22 00:00:00 UTC 2020
PD-L1 promotes tumor growth and progression by activating WIP and β-catenin signaling pathways and predicts poor prognosis in lung cancer.
Cell death & disease
Yu W,Hua Y,Qiu H,Hao J,Zou K,Li Z,Hu S,Guo P,Chen M,Sui S,Xiong Y,Li F,Lu J,Guo W,Luo G,Deng W
Mon Jul 06 00:00:00 UTC 2020
MicroRNA-410-3p Binds to TLR2 and Alleviates Myocardial Mitochondrial Dysfunction and Chemokine Production in LPS-Induced Sepsis.
Molecular therapy. Nucleic acids
Zuo T,Tang Q,Zhang X,Shang F
Fri Dec 04 00:00:00 UTC 2020
Regulatory coupling between long noncoding RNAs and senescence in irradiated microglia.
Journal of neuroinflammation
Xu A,Li R,Ren A,Jian H,Huang Z,Zeng Q,Wang B,Zheng J,Chen X,Zheng N,Zheng R,Tian Y,Liu M,Mao Z,Ji A,Yuan Y
Wed Oct 28 00:00:00 UTC 2020
miR-21a in exosomes from Lewis lung carcinoma cells accelerates tumor growth through targeting PDCD4 to enhance expansion of myeloid-derived suppressor cells.
Oncogene
Zhang X,Li F,Tang Y,Ren Q,Xiao B,Wan Y,Jiang S
Thu Oct 01 00:00:00 UTC 2020
Effects of cisplatin on the proliferation, invasion and apoptosis of breast cancer cells following β‑catenin silencing.
International journal of molecular medicine
Zhu X,Feng J,Fu W,Shu X,Wan X,Liu J
Mon Jun 01 00:00:00 UTC 2020
MiR-34a affects G2 arrest in prostate cancer PC3 cells via Wnt pathway and inhibits cell growth and migration.
European review for medical and pharmacological sciences
Dong B,Xu GC,Liu ST,Liu T,Geng B
Sat Aug 01 00:00:00 UTC 2020
Insulin secretion impairment induced by rosuvastatin partly though autophagy in INS-1E cells.
Cell biology international
Qian L,Zhu K,Lin Y,An L,Huang F,Yao Y,Ren L
Wed Jan 01 00:00:00 UTC 2020
miR-5089-5p suppresses castration-resistant prostate cancer resistance to enzalutamide and metastasis via miR-5089-5p/SPINK1/ MAPK/MMP9 signaling.
Aging
Wang ZC,Li Y,Wang KL,Wang L,You BS,Zhao DF,Liu ZQ,Fang RZ,Wang JQ,Zhang W,Zhang JM,Xu WH
Tue Jul 21 00:00:00 UTC 2020
Club cell 10-kDa protein (CC10) inhibits cPLA2/COX2 pathway to alleviate RSV-induced airway inflammation and AHR.
International immunopharmacology
Yu G,Mo S,Gao L,Wen X,Chen S,Long X,Xie X,Deng Y,Ren L,Zang N,Chen S,Liu E
Mon Jun 01 00:00:00 UTC 2020
Dendrobium Alkaloids Promote Neural Function After Cerebral Ischemia-Reperfusion Injury Through Inhibiting Pyroptosis Induced Neuronal Death in both In Vivo and In Vitro Models.
Neurochemical research
Liu D,Dong Z,Xiang F,Liu H,Wang Y,Wang Q,Rao J
Sat Feb 01 00:00:00 UTC 2020
MiR-22 restrains proliferation of rheumatoid arthritis by targeting IL6R and may be concerned with the suppression of NF-κB pathway.
The Kaohsiung journal of medical sciences
Yang QY,Yang KP,Li ZZ
Wed Jan 01 00:00:00 UTC 2020
Responses of intestinal morphology and function in offspring to heat stress in primiparous sows during late gestation.
Journal of thermal biology
Guo H,He J,Yang X,Zheng W,Yao W
Wed Apr 01 00:00:00 UTC 2020
Exosomes derived from smooth muscle cells ameliorate diabetes-induced erectile dysfunction by inhibiting fibrosis and modulating the NO/cGMP pathway.
Journal of cellular and molecular medicine
Song J,Sun T,Tang Z,Ruan Y,Liu K,Rao K,Lan R,Wang S,Wang T,Liu J
Sun Nov 01 00:00:00 UTC 2020
ZEB1-Mediated Transcriptional Upregulation of circWWC3 Promotes Breast Cancer Progression through Activating Ras Signaling Pathway.
Molecular therapy. Nucleic acids
Meng L,Liu S,Liu F,Sang M,Ju Y,Fan X,Gu L,Li Z,Geng C,Sang M
Wed Aug 19 00:00:00 UTC 2020
NNK-mediated upregulation of DEPDC1 stimulates the progression of oral squamous cell carcinoma by inhibiting CYP27B1 expression.
American journal of cancer research
Guo J,Zhou S,Huang P,Xu S,Zhang G,He H,Zeng Y,Xu CX,Kim H,Tan Y
Mon Sep 28 00:00:00 UTC 2020
Scavenger receptor A impairs interferon response to HBV infection by limiting TRAF3 ubiquitination through recruiting OTUB1.
The FEBS journal
Xie M,Yin Y,Chen L,Yin A,Liu Y,Liu Y,Dong L,Lai Q,Zhou J,Zhang L,Xu M,Chen Z,Zuo D
Wed Jan 01 00:00:00 UTC 2020
miRNA-182 regulated MTSS1 inhibits proliferation and invasion in Glioma Cells.
Journal of Cancer
Li Z,Zhang L,Liu Z,Huang T,Wang Y,Ma Y,Fang X,He Y,Zhou Y,Huo L,Wu J
Mon Sep 28 00:00:00 UTC 2020
BVES downregulation in non-syndromic tetralogy of fallot is associated with ventricular outflow tract stenosis.
Scientific reports
Shi Y,Li Y,Wang Y,Zhu P,Chen Y,Wang H,Yue S,Xia X,Chen J,Jiang Z,Zhou C,Cai W,Yuan H,Wu Y,Wan Y,Li X,Zhu X,Zhou Z,Dai G,Li F,Mo X,Ye X,Fan X,Zhuang J,Wu X,Yuan W
Tue Aug 25 00:00:00 UTC 2020
Characterization and Neuroprotection Potential of Seleno-Polymannuronate.
Frontiers in pharmacology
Bi D,Li X,Li T,Li X,Lin Z,Yao L,Li H,Xu H,Hu Z,Zhang Z,Liu Q,Xu X
Mon Sep 28 00:00:00 UTC 2020
Identification of EMT-related high-risk stage II colorectal cancer and characterisation of metastasis-related genes.
British journal of cancer
Wang K,Song K,Ma Z,Yao Y,Liu C,Yang J,Xiao H,Zhang J,Zhang Y,Zhao W
Sat Aug 01 00:00:00 UTC 2020
Ability of Post-treatment Glycyrrhizic Acid to Mitigate Cerebral Ischemia/Reperfusion Injury in Diabetic Mice.
Medical science monitor : international medical journal of experimental and clinical research
Li Y,Yao N,Zhang T,Guo F,Niu X,Wu Z,Hou S
Mon Sep 28 00:00:00 UTC 2020
Regulation of epidermal differentiation through KDF1-mediated deubiquitination of IKKα.
EMBO reports
Li Y,Tang L,Yue J,Gou X,Lin A,Weatherbee SD,Wu X
Wed May 06 00:00:00 UTC 2020
Human umbilical cord blood mononuclear cells protect against renal tubulointerstitial fibrosis in cisplatin-treated rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li XW,Feng LX,Zhu XJ,Liu Q,Wang HS,Wu X,Yan P,Duan XJ,Xiao YQ,Cheng W,Peng JC,Zhao F,Deng YH,Duan SB
Wed Jan 01 00:00:00 UTC 2020
CMIP promotes Herceptin resistance of HER2 positive gastric cancer cells.
Pathology, research and practice
Xiang R,Han X,Ding K,Wu Z
Sat Feb 01 00:00:00 UTC 2020
PRRX1-induced epithelial-to-mesenchymal transition in salivary adenoid cystic carcinoma activates the metabolic reprogramming of free fatty acids to promote invasion and metastasis.
Cell proliferation
Jiang YP,Tang YL,Wang SS,Wu JS,Zhang M,Pang X,Wu JB,Chen Y,Tang YJ,Liang XH
Wed Jan 01 00:00:00 UTC 2020
GALNT2 regulates ANGPTL3 cleavage in cells and in vivo of mice.
Scientific reports
Li X,Zhang Y,Zhang M,Wang Y
Wed Sep 30 00:00:00 UTC 2020
HIF-1α ameliorates tubular injury in diabetic nephropathy via HO-1-mediated control of mitochondrial dynamics.
Cell proliferation
Jiang N,Zhao H,Han Y,Li L,Xiong S,Zeng L,Xiao Y,Wei L,Xiong X,Gao P,Yang M,Liu Y,Sun L
Sun Nov 01 00:00:00 UTC 2020
Mitochondrial DNA Haplogroup M7 Confers Disability in a Chinese Aging Population.
Frontiers in genetics
Sun D,Yao S,Wu F,Deng W,Ma Y,Jin L,Wang J,Wang X
Tue Nov 17 00:00:00 UTC 2020
Lycopene attenuates body weight gain through induction of browning via regulation of peroxisome proliferator-activated receptor γ in high-fat diet-induced obese mice.
The Journal of nutritional biochemistry
Zhu R,Wei J,Liu H,Liu C,Wang L,Chen B,Li L,Jia Q,Tian Y,Li R,Zhao D,Mo F,Li Y,Gao S,Wang XD,Zhang D
Wed Apr 01 00:00:00 UTC 2020
NGF receptors and PI3K/AKT pathway involved in glucose fluctuation-induced damage to neurons and α-lipoic acid treatment.
BMC neuroscience
Yan T,Zhang Z,Li D
Thu Sep 17 00:00:00 UTC 2020
Metformin ameliorates cardiac conduction delay by regulating microRNA-1 in mice.
European journal of pharmacology
Lv L,Zheng N,Zhang L,Li R,Li Y,Yang R,Li C,Fang R,Shabanova A,Li X,Liu Y,Liang H,Zhou Y,Shan H
Sat Aug 15 00:00:00 UTC 2020
α-Ketoglutarate Modulates Macrophage Polarization Through Regulation of PPARγ Transcription and mTORC1/p70S6K Pathway to Ameliorate ALI/ARDS.
Shock (Augusta, Ga.)
Liu M,Chen Y,Wang S,Zhou H,Feng D,Wei J,Shi X,Wu L,Zhang P,Yang H,Lv X
Wed Jan 01 00:00:00 UTC 2020
Overexpression of endogenous lipoic acid synthase attenuates pulmonary fibrosis induced by crystalline silica in mice.
Toxicology letters
Zhao Y,Xu G,Li H,Chang M,Guan Y,Li Y,Wu W,Yao S
Fri May 01 00:00:00 UTC 2020
Deficiency of pseudogene UPAT leads to hepatocellular carcinoma progression and forms a positive feedback loop with ZEB1.
Cancer science
Xiang L,Huang X,Wang S,Ou H,Chen Z,Hu Z,Huang Y,Li X,Yuan Y,Yang D
Sun Nov 01 00:00:00 UTC 2020
Proteomics Analysis of Brain Tissue in a Rat Model of Ischemic Stroke in the Acute Phase.
Frontiers in molecular neuroscience
Zheng F,Zhou YT,Zeng YF,Liu T,Yang ZY,Tang T,Luo JK,Wang Y
Mon Sep 28 00:00:00 UTC 2020
Inhibition of miRNA-29a regulates intestinal barrier function in diarrhea-predominant irritable bowel syndrome by upregulating ZO-1 and CLDN1.
Experimental and therapeutic medicine
Zhu H,Xiao X,Shi Y,Wu Y,Huang Y,Li D,Xiong F,He G,Chai Y,Tang H
Tue Dec 01 00:00:00 UTC 2020
Honokiol inhibits the growth of SKBR3 cells.
Translational cancer research
Shi H,Wang Y,Yao M,Zhang D,Fang W,Zhou T,Gan D,Yue S,Qian H,Chen T
Tue Dec 01 00:00:00 UTC 2020
Silychristin A activates Nrf2-HO-1/SOD2 pathway to reduce apoptosis and improve GLP-1 production through upregulation of estrogen receptor α in GLUTag cells.
European journal of pharmacology
Wang J,Zhang X,Zhang L,Yan T,Wu B,Xu F,Jia Y
Sat Aug 15 00:00:00 UTC 2020
Lysine acetylation of F-actin decreases tropomyosin-based inhibition of actomyosin activity.
The Journal of biological chemistry
Schmidt W,Madan A,Foster DB,Cammarato A
Fri Nov 13 00:00:00 UTC 2020
Vitamin D3 protects articular cartilage by inhibiting the Wnt/β-catenin signaling pathway.
Experimental and therapeutic medicine
Yang T,Sun W,Duan YH,Sun YB,Ren YM,Hou WY,Tian MQ
Sat Aug 01 00:00:00 UTC 2020
Liraglutide Ameliorates Erectile Dysfunction via Regulating Oxidative Stress, the RhoA/ROCK Pathway and Autophagy in Diabetes Mellitus.
Frontiers in pharmacology
Yuan P,Ma D,Gao X,Wang J,Li R,Liu Z,Wang T,Wang S,Liu J,Liu X
Sat Apr 16 00:00:00 UTC 2022
Downregulation of HDAC1 suppresses media degeneration by inhibiting the migration and phenotypic switch of aortic vascular smooth muscle cells in aortic dissection.
Journal of cellular physiology
Sun L,Wang C,Yuan Y,Guo Z,He Y,Ma W,Zhang J
Sun Nov 01 00:00:00 UTC 2020
Oral recombinant methioninase increases TRAIL receptor-2 expression to regress pancreatic cancer in combination with agonist tigatuzumab in an orthotopic mouse model.
Cancer letters
Yamamoto J,Miyake K,Han Q,Tan Y,Inubushi S,Sugisawa N,Higuchi T,Tashiro Y,Nishino H,Homma Y,Matsuyama R,Chawla SP,Bouvet M,Singh SR,Endo I,Hoffman RM
Sun Nov 01 00:00:00 UTC 2020
Systematic investigation of biomarker-like role of ARHGDIB in breast cancer.
Cancer biomarkers : section A of Disease markers
Wang X,Bi X,Huang X,Wang B,Guo Q,Wu Z
Wed Feb 03 00:00:00 UTC 2021
Leucine-rich α-2-glycoprotein-1 promotes diabetic corneal epithelial wound healing and nerve regeneration via regulation of matrix metalloproteinases.
Experimental eye research
Li W,Wang X,Cheng J,Li J,Wang Q,Zhou Q,Li H,Xue J,Zhang Y,Yang L,Xie L
Wed Jul 01 00:00:00 UTC 2020
USP30 sets a trigger threshold for PINK1-PARKIN amplification of mitochondrial ubiquitylation.
Life science alliance
Rusilowicz-Jones EV,Jardine J,Kallinos A,Pinto-Fernandez A,Guenther F,Giurrandino M,Barone FG,McCarron K,Burke CJ,Murad A,Martinez A,Marcassa E,Gersch M,Buckmelter AJ,Kayser-Bricker KJ,Lamoliatte F,Gajbhiye A,Davis S,Scott HC,Murphy E,England K,Mortiboys H,Komander D,Trost M,Kessler BM,Ioannidis S,Ahlijanian MK,Urbé S,Clague MJ
Sat Aug 01 00:00:00 UTC 2020
Pretreatment with the ALDH2 activator Alda‑1 protects rat livers from ischemia/reperfusion injury by inducing autophagy.
Molecular medicine reports
Liu Z,Ye S,Zhong X,Wang W,Lai CH,Yang W,Yue P,Luo J,Huang X,Zhong Z,Xiong Y,Fan X,Li L,Wang Y,Ye Q
Tue Sep 01 00:00:00 UTC 2020
HIF-1α-upregulated lncRNA-H19 regulates lipid droplet metabolism through the AMPKα pathway in hepatic stellate cells.
Life sciences
Wang Z,Yang X,Kai J,Wang F,Wang Z,Shao J,Tan S,Chen A,Zhang F,Wang S,Zhang Z,Zheng S
Sat Aug 15 00:00:00 UTC 2020
Alda-1, an Aldehyde Dehydrogenase 2 Agonist, Improves Cutaneous Wound Healing by Activating Epidermal Keratinocytes via Akt/GSK-3β/β-Catenin Pathway.
Aesthetic plastic surgery
Zhang S,Chen C,Ying J,Wei C,Wang L,Yang Z,Qi F
Mon Jun 01 00:00:00 UTC 2020
Inhibition of Skp2 Sensitizes Chronic Myeloid Leukemia Cells to Imatinib.
Cancer management and research
Chen X,Huang Z,Wu W,Xia R
Fri Apr 15 00:00:00 UTC 2022
ROS enhances the cytotoxicity of cisplatin by inducing apoptosis and autophagy in tongue squamous cell carcinoma cells.
The international journal of biochemistry & cell biology
Xue DF,Pan ST,Huang G,Qiu JX
Fri May 01 00:00:00 UTC 2020
Spinosin Inhibits Aβ1-42 Production and Aggregation via Activating Nrf2/HO-1 Pathway.
Biomolecules & therapeutics
Zhang X,Wang J,Gong G,Ma R,Xu F,Yan T,Wu B,Jia Y
Fri May 01 00:00:00 UTC 2020
Ellagic Acid Protects Dopamine Neurons via Inhibition of NLRP3 Inflammasome Activation in Microglia.
Oxidative medicine and cellular longevity
He XM,Zhou YZ,Sheng S,Li JJ,Wang GQ,Zhang F
Tue Jun 08 00:00:00 UTC 2021
Novel ESCC-related gene ZNF750 as potential Prognostic biomarker and inhibits Epithelial-Mesenchymal Transition through directly depressing SNAI1 promoter in ESCC.
Theranostics
Kong P,Xu E,Bi Y,Xu X,Liu X,Song B,Zhang L,Cheng C,Yan T,Qian Y,Yang J,Ma Y,Cui H,Zhai Y,Zou B,Liu X,Cheng Y,Guo S,Cheng X,Cui Y
Tue Apr 13 00:00:00 UTC 2021
Novel mechanisms underlying anti-polycystic ovary like syndrome effects of electroacupuncture in rats: suppressing SREBP1 to mitigate insulin resistance, mitochondrial dysfunction and oxidative stress.
Biological research
Peng Y,Yang X,Luo X,Liu C,Cao X,Wang H,Guo L
Tue Oct 27 00:00:00 UTC 2020
Fenofibrate-induced mitochondrial dysfunction and metabolic reprogramming reversal: the anti-tumor effects in gastric carcinoma cells mediated by the PPAR pathway.
American journal of translational research
Chen L,Peng J,Wang Y,Jiang H,Wang W,Dai J,Tang M,Wei Y,Kuang H,Xu G,Xu H,Zhou F
Mon Sep 28 00:00:00 UTC 2020
CACNA1H downregulation induces skeletal muscle atrophy involving endoplasmic reticulum stress activation and autophagy flux blockade.
Cell death & disease
Li S,Hao M,Li B,Chen M,Chen J,Tang J,Hong S,Min J,Hu M,Hong L
Fri Apr 24 00:00:00 UTC 2020
Hsp90 inhibitor HSP990 in very low dose upregulates EAAT2 and exerts potent antiepileptic activity.
Theranostics
Sha L,Chen T,Deng Y,Du T,Ma K,Zhu W,Shen Y,Xu Q
Wed May 05 00:00:00 UTC 2021
Artesunate induces autophagy dependent apoptosis through upregulating ROS and activating AMPK-mTOR-ULK1 axis in human bladder cancer cells.
Chemico-biological interactions
Zhou X,Chen Y,Wang F,Wu H,Zhang Y,Liu J,Cai Y,Huang S,He N,Hu Z,Jin X
Sun Nov 01 00:00:00 UTC 2020
Utilizing methylglyoxal and D-lactate in urine to evaluate saikosaponin C treatment in mice with accelerated nephrotoxic serum nephritis.
PloS one
Lin CY,Lee JA,Lin PY,Hua SC,Tsai PY,Chen BL,Lin CE,Lee TH,Chen SM
Thu Dec 17 00:00:00 UTC 2020
MiR-145-5p alleviates hypoxia/reoxygenation- induced cardiac microvascular endothelial cell injury in coronary heart disease by inhibiting Smad4 expression.
European review for medical and pharmacological sciences
Li LL,Mao CD,Wang GP,Wang N,Xue AG
Fri May 01 00:00:00 UTC 2020
Progesterone Regulates Glucose Metabolism Through Glucose Transporter 1 to Promote Endometrial Receptivity.
Frontiers in physiology
Zhang H,Qi J,Wang Y,Sun J,Li Z,Sui L,Fan J,Liu C,Shang Y,Kong L,Kong Y
Wed Oct 28 00:00:00 UTC 2020
Microglia polarization of hippocampus is involved in the mechanism of Apelin-13 ameliorating chronic water immersion restraint stress-induced depression-like behavior in rats.
Neuropeptides
Zhou S,Chen S,Xie W,Guo X,Zhao J
Mon Jun 01 00:00:00 UTC 2020
Protective effect of Berberine on reproductive function and spermatogenesis in diabetic rats via inhibition of ROS/JAK2/NFκB pathway.
Andrology
Song J,Gao X,Tang Z,Li H,Ruan Y,Liu Z,Wang T,Wang S,Liu J,Jiang H
Fri May 01 00:00:00 UTC 2020
Stromal fibroblast-derived MFAP5 promotes the invasion and migration of breast cancer cells via Notch1/slug signaling.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico
Chen Z,Yan X,Li K,Ling Y,Kang H
Wed Apr 01 00:00:00 UTC 2020
MicroRNA-183 regulates lipopolysaccharide-induced oxidative stress of hippocampal neurons by targeting the fibronectin 1 gene.
Neuroscience letters
Xie Q,Feng L,Zhang C,Gan L
Thu Apr 23 00:00:00 UTC 2020
Differential distribution and potential regulatory roles of estrogen receptor 2a and 2b in the pituitary of ricefield eel Monopterus albus.
General and comparative endocrinology
Zheng Z,Liu M,Meng F,Zhang W,Zhang L
Sun Nov 01 00:00:00 UTC 2020
Hyperthermic intraperitoneal chemotherapy with recombinant mutant human TNF-α and raltitrexed in mice with colorectal-peritoneal carcinomatosis.
Experimental biology and medicine (Maywood, N.J.)
Gong Q,Song C,Wang X,Wang R,Cai G,Liang X,Liu J
Sun Mar 01 00:00:00 UTC 2020
MicroRNA-9 mediated the protective effect of ferulic acid on hypoxic-ischemic brain damage in neonatal rats.
PloS one
Yao K,Yang Q,Li Y,Lan T,Yu H,Yu Y
Fri Jul 24 00:00:00 UTC 2020
Role of the Nucleotide-Binding Domain-Like Receptor Protein 3 Inflammasome in the Endothelial Dysfunction of Early Sepsis.
Inflammation
Luo M,Meng J,Yan J,Shang F,Zhang T,Lv D,Li C,Yang X,Luo S
Sat Aug 01 00:00:00 UTC 2020
Odor-mediated contextual learning induced memory consolidation and hippocampus development in neonate rat.
Neuroreport
Cao J,Xu J,Chen S,Fan F,Chen M,Liu F,Li G
Wed Jan 08 00:00:00 UTC 2020
Echinacea polysaccharide alleviates LPS-induced lung injury via inhibiting inflammation, apoptosis and activation of the TLR4/NF-κB signal pathway.
International immunopharmacology
Zhang H,Lang W,Wang S,Li B,Li G,Shi Q
Sun Nov 01 00:00:00 UTC 2020
Pycnogenol achieves neuroprotective effects in rats with spinal cord injury by stabilizing the mitochondrial membrane potential.
Neurological research
Tao L,Liu X,Da W,Tao Z,Zhu Y
Wed Jul 01 00:00:00 UTC 2020
Anti-inflammation and anti-apoptosis effects of growth arrest-specific protein 6 in acute liver injury induced by LPS/D-GalN in mice.
Acta cirurgica brasileira
Wang Q,Zhao Y,Zang B
Fri May 01 00:00:00 UTC 2020
Computational repositioning of dimethyl fumarate for treating alcoholic liver disease.
Cell death & disease
Zhang Y,Zhao S,Fu Y,Yan L,Feng Y,Chen Y,Wu Y,Deng Y,Zhang G,Chen Z,Chen Y,Liu T
Tue Aug 18 00:00:00 UTC 2020
Progesterone promotes endothelial nitric oxide synthase expression through enhancing nuclear progesterone receptor-SP-1 formation.
American journal of physiology. Heart and circulatory physiology
You Y,Tan W,Guo Y,Luo M,Shang FF,Xia Y,Luo S
Sat Aug 01 00:00:00 UTC 2020
Fingolimod (FTY720) improves postoperative cognitive dysfunction in mice subjected to D-galactose-induced aging.
Neural regeneration research
Zhang J,Xiao B,Li CX,Wang Y
Wed Jul 01 00:00:00 UTC 2020
Effect of Combined Sorafenib/Cisplatinum Treatment on the Autophagy and Proliferation of Hepatocellular Carcinoma hepG2 Cells in Vitro.
Asian Pacific journal of cancer prevention : APJCP
Wang Y,Wang L
Thu Oct 01 00:00:00 UTC 2020
Hispidulin Inhibits Neuroinflammation in Lipopolysaccharide-Activated BV2 Microglia and Attenuates the Activation of Akt, NF-κB, and STAT3 Pathway.
Neurotoxicity research
Yu CI,Cheng CI,Kang YF,Chang PC,Lin IP,Kuo YH,Jhou AJ,Lin MY,Chen CY,Lee CH
Mon Jun 01 00:00:00 UTC 2020
A glycolysis-based 4-mRNA signature correlates with the prognosis and cell cycle process in patients with bladder cancer.
Cancer cell international
Zhang C,Gou X,He W,Yang H,Yin H
Thu Apr 14 00:00:00 UTC 2022
Functional Food XingJiuTang Attenuates Alcohol-Induced Liver Injury by Regulating SIRT1/Nrf-2 Signaling Pathway.
Chemistry & biodiversity
Hao J,Ding Y,Shi W,Zhang C,Li R
Tue Dec 01 00:00:00 UTC 2020
Prenatal hypoxia inhibited propionate-evoked BK channels of mesenteric artery smooth muscle cells in offspring.
Journal of cellular and molecular medicine
Zhang W,Feng X,Zhang Y,Sun M,Li L,Gao Q,Tang J,Zhang P,Lv J,Zhou X,Xu Z
Sun Mar 01 00:00:00 UTC 2020
Protective effects of Salidroside on spermatogenesis in streptozotocin induced type-1 diabetic male mice by inhibiting oxidative stress mediated blood-testis barrier damage.
Chemico-biological interactions
Jiang YP,Ye RJ,Yang JM,Liu N,Zhang WJ,Ma L,Sun T,Niu JG,Zheng P,Yu JQ
Sun Jan 05 00:00:00 UTC 2020
The Regulatory Mechanism and Biological Significance of Mitochondrial Calcium Uniporter in the Migration, Invasion, Angiogenesis and Growth of Gastric Cancer.
OncoTargets and therapy
Wang X,Song X,Cheng G,Zhang J,Dong L,Bai J,Luo D,Xiong Y,Li S,Liu F,Sun Y,Wang X,Li Y,Huang Y
Mon Apr 18 00:00:00 UTC 2022
RNA N6-methyladenosine reader IGF2BP3 regulates cell cycle and angiogenesis in colon cancer.
Journal of experimental & clinical cancer research : CR
Yang Z,Wang T,Wu D,Min Z,Tan J,Yu B
Tue Sep 29 00:00:00 UTC 2020
Protein microarray analysis of cytokine expression changes in distal stumps after sciatic nerve transection.
Neural regeneration research
Cheng XQ,Liang XZ,Wei S,Ding X,Han GH,Liu P,Sun X,Quan Q,Tang H,Zhao Q,Shang AJ,Peng J
Sun Mar 01 00:00:00 UTC 2020
Aspirin enhances the sensitivity of colon cancer cells to cisplatin by abrogating the binding of NF-κB to the COX-2 promoter.
Aging
Jiang W,Yan Y,Chen M,Luo G,Hao J,Pan J,Hu S,Guo P,Li W,Wang R,Zuo Y,Sun Y,Sui S,Yu W,Pan Z,Zou K,Zheng Z,Deng W,Wu X,Guo W
Mon Jan 06 00:00:00 UTC 2020
MG53 Protects hUC-MSCs against Inflammatory Damage and Synergistically Enhances Their Efficacy in Neuroinflammation Injured Brain through Inhibiting NLRP3/Caspase-1/IL-1β Axis.
ACS chemical neuroscience
Ma S,Wang Y,Zhou X,Li Z,Zhang Z,Wang Y,Huang T,Zhang Y,Shi J,Guan F
Wed Sep 02 00:00:00 UTC 2020
Aberrant BMP2 Signaling in Patients Diagnosed with Osteoporosis.
International journal of molecular sciences
Durbano HW,Halloran D,Nguyen J,Stone V,McTague S,Eskander M,Nohe A
Mon Sep 21 00:00:00 UTC 2020
Nicotinamide Riboside Enhances Mitochondrial Proteostasis and Adult Neurogenesis through Activation of Mitochondrial Unfolded Protein Response Signaling in the Brain of ALS SOD1G93A Mice.
International journal of biological sciences
Zhou Q,Zhu L,Qiu W,Liu Y,Yang F,Chen W,Xu R
Mon Nov 02 00:00:00 UTC 2020
Methylglyoxal and D-lactate in cisplatin-induced acute kidney injury: Investigation of the potential mechanism via fluorogenic derivatization liquid chromatography-tandem mass spectrometry (FD-LC-MS/MS) proteomic analysis.
PloS one
Chen SM,Chen TH,Chang HT,Lin TY,Lin CY,Tsai PY,Imai K,Chen CM,Lee JA
Mon Sep 14 00:00:00 UTC 2020
Association analysis of DNA methyltransferases in IgA nephropathy.
International immunopharmacology
Xia M,Chen G,Liu D,Tang X,Liu Y,Wu L,Liu Y,Liu H
Sun Mar 01 00:00:00 UTC 2020
Dihydroartemisinin Prevents Progression and Metastasis of Head and Neck Squamous Cell Carcinoma by Inhibiting Polarization of Macrophages in Tumor Microenvironment.
OncoTargets and therapy
Chen R,Lu X,Li Z,Sun Y,He Z,Li X
Thu Apr 14 00:00:00 UTC 2022
Weighted gene co-expression network analysis identifies CCNA2 as a treatment target of prostate cancer through inhibiting cell cycle.
Journal of Cancer
Yang R,Du Y,Wang L,Chen Z,Liu X
Wed Sep 30 00:00:00 UTC 2020
Transgelin-2 contributes to proliferation and progression of hepatocellular carcinoma via regulating Annexin A2.
Biochemical and biophysical research communications
Shi J,Ren M,She X,Zhang Z,Zhao Y,Han Y,Lu D,Lyu L
Thu Mar 12 00:00:00 UTC 2020
High glucose inhibits vascular endothelial Keap1/Nrf2/ARE signal pathway via downregulation of monomethyltransferase SET8 expression.
Acta biochimica et biophysica Sinica
Chen X,Qi J,Wu Q,Jiang H,Wang J,Chen W,Mao A,Zhu M
Tue May 26 00:00:00 UTC 2020
Compositional characterization of Pyrus ussuriensis Maxim and their antioxidant activities and induction of apoptosis in Bel-7402 cell.
Journal of food biochemistry
Peng F,Li G,Xie Y,Yin HY,Li XJ,Yang YD
Mon Jun 01 00:00:00 UTC 2020
SLC25A10 performs an oncogenic role in human osteosarcoma.
Oncology letters
Wang G,Xia J,Chen C,Qiu J,Sun P,Peng Z,Chen X,Xu B
Thu Oct 01 00:00:00 UTC 2020
Antagomirs targeting miR-142-5p attenuate pilocarpine-induced status epilepticus in mice.
Experimental cell research
Zhang H,Lian Y,Xie N,Cheng X,Chen C,Xu H,Zheng Y
Sat Aug 15 00:00:00 UTC 2020
Effects of Hydrogen-rich Water on the PI3K/AKT Signaling Pathway in Rats with Myocardial Ischemia-reperfusion Injury.
Current molecular medicine
Li L,Li X,Zhang Z,Liu L,Liu T,Li S,Liu S,Zhou Y,Liu F
Wed Jun 09 00:00:00 UTC 2021
Dihydromethysticin, a natural molecule from Kava, suppresses the growth of colorectal cancer via the NLRC3/PI3K pathway.
Molecular carcinogenesis
Pan H,Liu F,Wang J,Zhao M,Wang D,Jia C,Wang T,Chen Z,Fan Y,Liang D,Meng Q
Mon Jun 01 00:00:00 UTC 2020
LncRNA MALAT1 knockdown inhibits cell migration and invasion by suppressing autophagy through miR-384/GOLM1 axis in glioma.
European review for medical and pharmacological sciences
Ma R,Zhang BW,Zhang ZB,Deng QJ
Sun Mar 01 00:00:00 UTC 2020
Establishment and characterization of an immortalized human chondrocyte cell line.
Biotechnology letters
Yang J,Tang Y,Chen W,Zhang M,Hong F,Yang D,Bao Q,Yang Y,Jin Y,Li L,Ma C
Fri May 01 00:00:00 UTC 2020
Identification of FADS1 Through Common Gene Expression Profiles for Predicting Survival in Patients with Bladder Cancer.
Cancer management and research
Jiao F,Sun H,Yang Q,Sun H,Wang Z,Liu M,Chen J
Sun Apr 17 00:00:00 UTC 2022
Nrf2 activation protects auditory hair cells from cisplatin-induced ototoxicity independent on mitochondrial ROS production.
Toxicology letters
Zhang W,Xiong H,Pang J,Su Z,Lai L,Lin H,Jian B,He W,Yang H,Zheng Y
Thu Oct 01 00:00:00 UTC 2020
Curcumin inhibits the growth of triple-negative breast cancer cells by silencing EZH2 and restoring DLC1 expression.
Journal of cellular and molecular medicine
Zhou X,Jiao D,Dou M,Zhang W,Lv L,Chen J,Li L,Wang L,Han X
Tue Sep 01 00:00:00 UTC 2020
Metformin alleviates breast cancer through targeting high-mobility group AT-hook 2.
Thoracic cancer
Li Y,Wang D,Ren H,Feng W
Sun Mar 01 00:00:00 UTC 2020
Polysaccharides from Armillariella tabescens mycelia ameliorate insulin resistance in type 2 diabetic mice.
Food & function
Yang R,Li Y,Cai J,Ji J,Wang Y,Zhang W,Pan W,Chen Y
Wed Nov 18 00:00:00 UTC 2020
LincRNA00494 Suppresses Non-small Cell Lung Cancer Cell Proliferation by Regulating SRCIN1 Expression as a ceRNA.
Frontiers in oncology
Dong J,Li B,Lin D,Lu D,Liu C,Lu X,Tang X,Li L,Zhu D,Liu J,Qiu X,Tian L,Zhou Q
Mon Sep 28 00:00:00 UTC 2020
Silencing ARAF Suppresses the Malignant Phenotypes of Gallbladder Cancer Cells.
BioMed research international
Lin W,Tong C,Zhang W,Cen W,Wang Y,Li J,Zhu Z,Yu J,Lu B
Thu Apr 29 00:00:00 UTC 2021
Role of Nrf2 in Lipopolysaccharide-Induced Acute Kidney Injury: Protection by Human Umbilical Cord Blood Mononuclear Cells.
Oxidative medicine and cellular longevity
Feng LX,Zhao F,Liu Q,Peng JC,Duan XJ,Yan P,Wu X,Wang HS,Deng YH,Duan SB
Fri May 14 00:00:00 UTC 2021
Curcumin protects rat hippocampal neurons against pseudorabies virus by regulating the BDNF/TrkB pathway.
Scientific reports
Yang B,Luo G,Zhang C,Feng L,Luo X,Gan L
Thu Dec 17 00:00:00 UTC 2020
Induction of SPARC on Oxidative Stress, Inflammatory Phenotype Transformation, and Apoptosis of Human Brain Smooth Muscle Cells Via TGF-β1-NOX4 Pathway.
Journal of molecular neuroscience : MN
Tan X,Li T,Zhu S,Zhong W,Li F,Wang Y
Sun Nov 01 00:00:00 UTC 2020
YBX1 mediates autophagy by targeting p110β and decreasing the sensitivity to cisplatin in NSCLC.
Cell death & disease
Cui Y,Li F,Xie Q,Zhao S,Guo T,Guo P,Hu S,Hao J,Tian C,Yu W,Li Z,Fang L,Zhao L,Chen M,Wu T,Gu C
Fri Jun 19 00:00:00 UTC 2020
Neuronostatin Promotion Soluble Aβ1-42 Oligomers: Induced Dysfunctional Brain Glucose Metabolism in Mice.
Neurochemical research
Yang S,Zhou F,Ma M,Yuan Y,Zhao S,Yu P
Thu Oct 01 00:00:00 UTC 2020
Indoxyl sulfate promotes osteogenic differentiation of vascular smooth muscle cells by miR-155-5p-dependent downregulation of matrix Gla protein via ROS/NF-κB signaling.
Experimental cell research
He X,Wang Z,Wei L,Cheng X,Chen L,Gao F,Jiang H
Tue Dec 01 00:00:00 UTC 2020
The Novel Synthetic Triterpene Methyl 3β-O-[4-(2-Aminoethylamino)-4-oxo-butyryl]olean-12-ene-28-oate Inhibits Breast Tumor Cell Growth in Vitro and in Vivo.
Chemical & pharmaceutical bulletin
Feng B,Zhao C,Li J,Yu J,Zhang Y,Zhang X,Tian T,Zhao L
Fri Apr 30 00:00:00 UTC 2021
Panax notoginseng saponin R1 modulates TNF-α/NF-κB signaling and attenuates allergic airway inflammation in asthma.
International immunopharmacology
Xue K,Ruan L,Hu J,Fu Z,Tian D,Zou W
Sun Nov 01 00:00:00 UTC 2020
Melatonin ameliorates hyperglycaemia-induced renal inflammation by inhibiting the activation of TLR4 and TGF-β1/Smad3 signalling pathway.
American journal of translational research
Wei J,Wang Y,Qi X,Fan Z,Wu Y
Mon Sep 28 00:00:00 UTC 2020
MTFP1 overexpression promotes the growth of oral squamous cell carcinoma by inducing ROS production.
Cell biology international
Xiao T,Sun J,Xing Z,Xie F,Yang L,Ding W
Sun Mar 01 00:00:00 UTC 2020
Euscaphic acid and Tormentic acid protect vascular endothelial cells against hypoxia-induced apoptosis via PI3K/AKT or ERK 1/2 signaling pathway.
Life sciences
Shi C,Li Z,Wu Y,Li X,Li Y,Wei J,Li J,Zhang Y,Li L
Wed Jul 01 00:00:00 UTC 2020
An autoregulatory feedback loop of miR-21/VMP1 is responsible for the abnormal expression of miR-21 in colorectal cancer cells.
Cell death & disease
Wang C,Peng R,Zeng M,Zhang Z,Liu S,Jiang D,Lu Y,Zou F
Mon Dec 14 00:00:00 UTC 2020
BCCIPβ facilitates p53 ubiquitination via binding with E6 protein in high-risk HPV positive head and neck squamous cell carcinoma.
Biochemical and biophysical research communications
Wang F,Wang J,Wang J,Zhang L,Fu H,Li J,Tian T,Zuo J,Lv W,Ma X
Thu Aug 27 00:00:00 UTC 2020
Identification of Immune-Related Genes Contributing to the Development of Glioblastoma Using Weighted Gene Co-expression Network Analysis.
Frontiers in immunology
Kong Y,Feng ZC,Zhang YL,Liu XF,Ma Y,Zhao ZM,Huang B,Chen AJ,Zhang D,Thorsen F,Wang J,Yang N,Li XG
Mon Apr 12 00:00:00 UTC 2021
Promotion of cadmium uptake and cadmium-induced toxicity by the copper transporter CTR1 in HepG2 and ZFL cells.
Toxicology reports
Kwok ML,Li ZP,Law TYS,Chan KM
Thu Dec 10 00:00:00 UTC 2020
Expression of lncRNA-HOTAIR in the serum of patients with lymph node metastasis of papillary thyroid carcinoma and its impact.
Oncology letters
Wu L,Shi Y,Liu B,Zhao M
Wed Jul 01 00:00:00 UTC 2020
Baicalin Inhibits Coxsackievirus B3 Replication by Reducing Cellular Lipid Synthesis.
The American journal of Chinese medicine
Wang MJ,Yang CH,Jin Y,Wan CB,Qian WH,Xing F,Li X,Liu YY
Fri Sep 04 00:00:00 UTC 2020
Decreased basal and stimulated salivary parameters by histopathological lesions and secretory dysfunction of parotid and submandibular glands in rats with type 2 diabetes.
Experimental and therapeutic medicine
Chen SY,Wang Y,Zhang CL,Yang ZM
Wed Apr 01 00:00:00 UTC 2020
Geniposide in Gardenia jasminoides var. radicans Makino modulates blood pressure via inhibiting WNK pathway mediated by the estrogen receptors.
The Journal of pharmacy and pharmacology
Fu Y,Yuan PP,Cao YG,Ke YY,Zhang Q,Hou Y,Zhang YL,Feng WS,Zheng XK
Tue Dec 01 00:00:00 UTC 2020
The testis-specific gene 1700102P08Rik is essential for male fertility.
Molecular reproduction and development
Wu XL,Yun DM,Gao S,Liang AJ,Duan ZZ,Wang HS,Wang GS,Sun F
Sat Feb 01 00:00:00 UTC 2020
Melatonin Ameliorates Renal Fibrosis Through the Inhibition of NF-κB and TGF-β1/Smad3 Pathways in db/db Diabetic Mice.
Archives of medical research
Fan Z,Qi X,Yang W,Xia L,Wu Y
Sat Aug 01 00:00:00 UTC 2020
Anti-insulin resistance effects of salidroside through mitochondrial quality control.
The Journal of endocrinology
You B,Dun Y,Zhang W,Jiang L,Li H,Xie M,Liu Y,Liu S
Sat Feb 01 00:00:00 UTC 2020
Effect of NF-κB signal pathway on mucus secretion induced by atmospheric PM2.5 in asthmatic rats.
Ecotoxicology and environmental safety
Liu Y,Zhang B,Zhang T,Wang H,Peng L,Zhou L
Sun Mar 01 00:00:00 UTC 2020
Inhibition of calcineurin/NFATc4 signaling attenuates ventilator‑induced lung injury.
Molecular medicine reports
Li M,Fang XZ,Liu XT,Zheng YF,Xie YB,Ma XD,Xia Y,Shao DH
Sat Feb 01 00:00:00 UTC 2020
Effect of autophagy on irradiation‑induced damage in osteoblast‑like MC3T3‑E1 cells.
Molecular medicine reports
Li R,Yang W,Hu X,Zhou D,Huang K,Wang C,Li Y,Liu B
Thu Oct 01 00:00:00 UTC 2020
Effect of KIF22 on promoting proliferation and migration of gastric cancer cells via MAPK-ERK pathways.
Chinese medical journal
Yu ZY,Jiang XY,Zhao RR,Qin JJ,Luo CJ,Ren YX,Ren W,Ma ZJ,Jiao ZY
Mon Apr 20 00:00:00 UTC 2020
Neuroprotective effects of notoginsenoside R1 by upregulating Trx-1 on acrylamide-induced neurotoxicity in PC12.
Human & experimental toxicology
Wang W,Huang L,Hu Y,Thomas ER,Li X
Mon Jun 01 00:00:00 UTC 2020
Activation of the Hedgehog Pathway Promotes Recovery of Neurological Function After Traumatic Brain Injury by Protecting the Neurovascular Unit.
Translational stroke research
Wu J,He J,Tian X,Zhong J,Li H,Sun X
Sat Aug 01 00:00:00 UTC 2020
The RBP1-CKAP4 axis activates oncogenic autophagy and promotes cancer progression in oral squamous cell carcinoma.
Cell death & disease
Gao L,Wang Q,Ren W,Zheng J,Li S,Dou Z,Kong X,Liang X,Zhi K
Thu Jun 25 00:00:00 UTC 2020
Ablation of glutaredoxin 1 promotes pulmonary angiogenesis and alveolar formation in hyperoxia-injured lungs by modifying HIF-1α stability and inhibiting the NF-κB pathway.
Biochemical and biophysical research communications
Liu X,Li K,Zhang F,Zhang Y,Deng C,Guo C
Thu Apr 30 00:00:00 UTC 2020
4-Acetylantroquinonol B Inhibits Osteoclastogenesis by Inhibiting the Autophagy Pathway in a Simulated Microgravity Model.
International journal of molecular sciences
Wu CH,Ou CH,Yen IC,Lee SY
Tue Sep 22 00:00:00 UTC 2020
Berberine alleviates cisplatin-induced acute kidney injury by regulating mitophagy via PINK 1/Parkin pathway.
Translational andrology and urology
Qi J,Xue Q,Kuang L,Xie L,Luo R,Nie X
Sat Aug 01 00:00:00 UTC 2020
Minicircle DNA vector expressing interferon-lambda-3 inhibits hepatitis B virus replication and expression in hepatocyte-derived cell line.
BMC molecular and cell biology
Guo X,Chen D,Cai Q,Huang Z,Xu W,Peng L,Chen P
Tue Feb 18 00:00:00 UTC 2020
Genkwadaphnin inhibits growth and invasion in hepatocellular carcinoma by blocking DHCR24-mediated cholesterol biosynthesis and lipid rafts formation.
British journal of cancer
Wu J,Guo L,Qiu X,Ren Y,Li F,Cui W,Song S
Sun Nov 01 00:00:00 UTC 2020
Poldip2 mediates blood-brain barrier disruption and cerebral edema by inducing AQP4 polarity loss in mouse bacterial meningitis model.
CNS neuroscience & therapeutics
Gao M,Lu W,Shu Y,Yang Z,Sun S,Xu J,Gan S,Zhu S,Qiu G,Zhuo F,Xu S,Wang Y,Chen J,Wu X,Huang J
Tue Dec 01 00:00:00 UTC 2020
Selaginellin B induces apoptosis and autophagy in pancreatic cancer cells via the JAK2/STAT3 signaling pathway.
American journal of translational research
Chu P,Wang S,Zhu X,Yang Y,Li H,Tesfaldet T,Shopit A,Yang Y,Ma X,Peng J,Tang Z,Sun Z
Wed Dec 16 00:00:00 UTC 2020
miR-100-3p inhibits the adipogenic differentiation of hMSCs by targeting PIK3R1 via the PI3K/AKT signaling pathway.
Aging
Wang T,Zhong D,Qin Z,He S,Gong Y,Li W,Li X
Fri Nov 20 00:00:00 UTC 2020
Novel nortriterpenoids with new skeletons and limonoids from the fruits of Evodia rutaecarpa and their bioactivities.
Fitoterapia
Shi YS,Xia HM,Wu CH,Li CB,Duan CC,Che C,Zhang XJ,Li HT,Zhang Y,Zhang XF
Wed Apr 01 00:00:00 UTC 2020
Endoplasmic reticulum stress remodels alveolar bone formation after tooth extraction.
Journal of cellular and molecular medicine
Chen Y,Guo Y,Li J,Chen YY,Liu Q,Tan L,Gao ZR,Zhang SH,Zhou YH,Feng YZ
Sun Nov 01 00:00:00 UTC 2020
Inhibition of the NLRP3-inflammasome prevents cognitive deficits in experimental autoimmune encephalomyelitis mice via the alteration of astrocyte phenotype.
Cell death & disease
Hou B,Zhang Y,Liang P,He Y,Peng B,Liu W,Han S,Yin J,He X
Fri May 15 00:00:00 UTC 2020
Phloretin Modulates Human Th17/Treg Cell Differentiation In Vitro via AMPK Signaling.
BioMed research international
Jiao A,Yang Z,Fu X,Hua X
Thu Apr 22 00:00:00 UTC 2021
Drp1, a potential therapeutic target for Parkinson's disease, is involved in olfactory bulb pathological alteration in the Rotenone-induced rat model.
Toxicology letters
Zhang X,Huang W,Shao Q,Yang Y,Xu Z,Chen J,Zhang X,Ge X
Mon Jun 01 00:00:00 UTC 2020
Cytisine attenuates bone loss of ovariectomy mouse by preventing RANKL-induced osteoclastogenesis.
Journal of cellular and molecular medicine
Qian Z,Zhong Z,Ni S,Li D,Zhang F,Zhou Y,Kang Z,Qian J,Yu B
Tue Sep 01 00:00:00 UTC 2020
The LINC00365/SCGB2A1 (Mammaglobin B) Axis Down-Regulates NF-κB Signaling and Is Associated with the Progression of Gastric Cancer.
Cancer management and research
Yan XY,Zhang JJ,Zhong XR,Yu SH,Xu L,Tian R,Sun LK,Su J
Mon Nov 13 00:00:00 UTC 2023
MicroRNA-145 suppresses osteogenic differentiation of human jaw bone marrow mesenchymal stem cells partially via targeting semaphorin 3A.
Connective tissue research
Jin Y,Hong F,Bao Q,Xu Q,Duan R,Zhu Z,Zhang W,Ma C
Sun Nov 01 00:00:00 UTC 2020
Beneficial Effects of Theta-Burst Transcranial Magnetic Stimulation on Stroke Injury via Improving Neuronal Microenvironment and Mitochondrial Integrity.
Translational stroke research
Zong X,Dong Y,Li Y,Yang L,Li Y,Yang B,Tucker L,Zhao N,Brann DW,Yan X,Hu S,Zhang Q
Mon Jun 01 00:00:00 UTC 2020
Hepatoprotective effects of Di Wu Yang Gan: A medicinal food against CCl4-induced hepatotoxicity in vivo and in vitro.
Food chemistry
Xu W,Xiao M,Li J,Chen Y,Sun Q,Li H,Sun W
Thu Oct 15 00:00:00 UTC 2020
Calcium-sensing receptor promotes tumor proliferation and migration in human intrahepatic cholangiocarcinoma by targeting ERK signaling pathway.
European journal of pharmacology
Fang Y,Liu L,Liu S,Hu L,Cai W,Wan X,Liu D,He Y,Zhu Z
Sun Apr 05 00:00:00 UTC 2020
Graphene oxide size-dependently altered lipid profiles in THP-1 macrophages.
Ecotoxicology and environmental safety
Luo Y,Peng J,Huang C,Cao Y
Sat Aug 01 00:00:00 UTC 2020
Leptin mediates protection of hydrogen sulfide against 6-hydroxydopamine-induced Parkinson's disease: Involving enhancement in Warburg effect.
Neurochemistry international
Yang SQ,Tian Q,Li D,He SQ,Hu M,Liu SY,Zou W,Chen YJ,Zhang P,Tang XQ
Fri May 01 00:00:00 UTC 2020
circFOXM1 promotes proliferation of non-small cell lung carcinoma cells by acting as a ceRNA to upregulate FAM83D.
Journal of experimental & clinical cancer research : CR
Yu C,Cheng Z,Cui S,Mao X,Li B,Fu Y,Wang H,Jin H,Ye Q,Zhao X,Jiang L,Qin W
Mon Mar 30 00:00:00 UTC 2020
High- and low-virulent bovine Pasteurella multocida induced differential NLRP3 inflammasome activation and subsequent IL-1β secretion.
Veterinary microbiology
Fang R,Lei G,Jiang J,Du H,Liu Y,Lei Z,Ye C,Li N,Peng Y
Wed Apr 01 00:00:00 UTC 2020
DUSP5 (dual-specificity protein phosphatase 5) suppresses BCG-induced autophagy via ERK 1/2 signaling pathway.
Molecular immunology
Luo J,Xue D,Song F,Liu X,Li W,Wang Y
Thu Oct 01 00:00:00 UTC 2020
Connective tissue growth factor as an unfavorable prognostic marker promotes the proliferation, migration, and invasion of gliomas.
Chinese medical journal
Song ZB,Yang HP,Xu AQ,Zhan ZM,Song Y,Li ZY
Fri Mar 20 00:00:00 UTC 2020
Hydrogen Sulfide Attenuates High-Fat Diet-Induced Non-Alcoholic Fatty Liver Disease by Inhibiting Apoptosis and Promoting Autophagy via Reactive Oxygen Species/Phosphatidylinositol 3-Kinase/AKT/Mammalian Target of Rapamycin Signaling Pathway.
Frontiers in pharmacology
Wu D,Zhong P,Wang Y,Zhang Q,Li J,Liu Z,Ji A,Li Y
Tue Jan 05 00:00:00 UTC 2021
Electroacupuncture Pretreatment Alleviates LPS-Induced Acute Respiratory Distress Syndrome via Regulating the PPAR Gamma/NF-Kappa B Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Feng D,Zhou H,Jin X,Wei J,Zhang Q,Gu Y,Zhang P,Yang H,Song J,Shi X,Lv X
Mon Sep 28 00:00:00 UTC 2020
Protective effects of essential oil from Fructus Alpiniae zerumbet on retinal Müller gliosis via the PPAR-γ-p-CREB signaling pathway.
Chinese medicine
Yang H,Gan S,Jiang Z,Song X,Chen T,Xu Y,Fu L,Zhang Y,Tao L,Shen X
Tue Apr 12 00:00:00 UTC 2022
MicroRNA 885-5p Inhibits Hepatocellular Carcinoma Metastasis by Repressing AEG1.
OncoTargets and therapy
Li C,Wang X,Song Q
Wed Apr 13 00:00:00 UTC 2022
REG γ knockdown suppresses proliferation by inducing apoptosis and cell cycle arrest in osteosarcoma.
PeerJ
Yin Z,Jin H,Huang S,Qu G,Meng Q
Thu Apr 14 00:00:00 UTC 2022
SMAD3/SP1 complex-mediated constitutive active loop between lncRNA PCAT7 and TGF-β signaling promotes prostate cancer bone metastasis.
Molecular oncology
Lang C,Dai Y,Wu Z,Yang Q,He S,Zhang X,Guo W,Lai Y,Du H,Wang H,Ren D,Peng X
Wed Apr 01 00:00:00 UTC 2020
Isorhamnetin attenuates Streptococcus suis virulence by inhibiting the inflammatory response.
Antonie van Leeuwenhoek
Li G,Wang G,Wang S,Sun M,Wen Z
Sat Feb 01 00:00:00 UTC 2020
The mechanism and role of intracellular α-ketoglutarate reduction in hepatic stellate cell activation.
Bioscience reports
Zhao J,Jiang Y,Sun X,Liu X,Liu F,Song M,Zhang L
Fri Mar 27 00:00:00 UTC 2020
Impaired mitochondrial medium-chain fatty acid oxidation drives periportal macrovesicular steatosis in sirtuin-5 knockout mice.
Scientific reports
Goetzman ES,Bharathi SS,Zhang Y,Zhao XJ,Dobrowolski SF,Peasley K,Sims-Lucas S,Monga SP
Tue Oct 27 00:00:00 UTC 2020
Network Pharmacology and Bioactive Equivalence Assessment Integrated Strategy Driven Q-markers Discovery for Da-Cheng-Qi Decoction to Attenuate Intestinal Obstruction.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li D,Lv B,Wang D,Xu D,Qin S,Zhang Y,Chen J,Zhang W,Zhang Z,Xu F
Wed Jul 01 00:00:00 UTC 2020
LINC00963 Promotes Ovarian Cancer Proliferation, Migration and EMT via the miR-378g /CHI3L1 Axis.
Cancer management and research
Liu W,Yang YJ,An Q
Mon Jan 29 00:00:00 UTC 2024
The Late Domain of Prototype Foamy Virus Gag Facilitates Autophagic Clearance of Stress Granules by Promoting Amphisome Formation.
Journal of virology
Zheng Y,Zhu G,Yan J,Tang Y,Han S,Yin J,Peng B,He X,Liu W
Tue Mar 17 00:00:00 UTC 2020
NRSN2 promotes breast cancer metastasis by activating PI3K/AKT/mTOR and NF-κB signaling pathways.
Oncology letters
Ren F,Zhang W,Lu S,Ren H,Guo Y
Wed Jan 01 00:00:00 UTC 2020
Resveratrol slows the tumourigenesis of pancreatic cancer by inhibiting NFκB activation.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Qian W,Xiao Q,Wang L,Qin T,Xiao Y,Li J,Yue Y,Zhou C,Duan W,Ma Q,Ma J
Wed Jul 01 00:00:00 UTC 2020
ROS-mediated PERK-eIF2α-ATF4 pathway plays an important role in arsenite-induced L-02 cells apoptosis via regulating CHOP-DR5 signaling.
Environmental toxicology
Liu C,Zhang A
Thu Oct 01 00:00:00 UTC 2020
Effects of inhalation of sevoflurane at different concentrations on TRPV1 in airways of rats at different developmental stages.
Life sciences
Liu D,Yuan J,Fei X,Zhu Y,Zhou Y,Zhang C,Dong L,Zhu Z
Fri May 15 00:00:00 UTC 2020
Blocking circ_0013912 Suppressed Cell Growth, Migration and Invasion of Pancreatic Ductal Adenocarcinoma Cells in vitro and in vivo Partially Through Sponging miR-7-5p.
Cancer management and research
Guo W,Zhao L,Wei G,Liu P,Zhang Y,Fu L
Sat Apr 16 00:00:00 UTC 2022
Romo1 is involved in the immune response of glioblastoma by regulating the function of macrophages.
Aging
Sun G,Cao Y,Qian C,Wan Z,Zhu J,Guo J,Shi L
Thu Jan 16 00:00:00 UTC 2020
MicroRNA-217 ameliorates inflammatory damage of endothelial cells induced by oxidized LDL by targeting EGR1.
Molecular and cellular biochemistry
Yang X,Li D,Qi YZ,Chen W,Yang CH,Jiang YH
Tue Dec 01 00:00:00 UTC 2020
Histone Deacetylation 10 Alleviates Inflammation After Intracerebral Hemorrhage via the PTPN22/NLRP3 Pathway in Rats.
Neuroscience
Wang L,Zheng S,Zhang L,Xiao H,Gan H,Chen H,Zhai X,Liang P,Zhao J,Li Y
Wed Apr 15 00:00:00 UTC 2020
Fisetin Alleviated Bleomycin-Induced Pulmonary Fibrosis Partly by Rescuing Alveolar Epithelial Cells From Senescence.
Frontiers in pharmacology
Zhang L,Tong X,Huang J,Wu M,Zhang S,Wang D,Liu S,Fan H
Fri Jan 01 00:00:00 UTC 2021
Alprostadil attenuates LPS-induced cardiomyocyte injury by inhibiting the Wnt5a/JNK/NF-κB pathway.
Herz
Yu T,Dong D,Guan J,Sun J,Guo M,Wang Q
Tue Dec 01 00:00:00 UTC 2020
Liver Expression of IGF2 and Related Proteins in ZBED6 Gene-Edited Pig by RNA-Seq.
Animals : an open access journal from MDPI
Zhao H,Wu M,Liu S,Tang X,Yi X,Li Q,Wang S,Sun X
Sun Nov 22 00:00:00 UTC 2020
Transforming growth factor beta type 1 (TGF-β) and hypoxia-inducible factor 1 (HIF-1) transcription complex as master regulators of the immunosuppressive protein galectin-9 expression in human cancer and embryonic cells.
Aging
Selnø ATH,Schlichtner S,Yasinska IM,Sakhnevych SS,Fiedler W,Wellbrock J,Klenova E,Pavlova L,Gibbs BF,Degen M,Schnyder I,Aliu N,Berger SM,Fasler-Kan E,Sumbayev VV
Tue Dec 08 00:00:00 UTC 2020
Amino Acids Regulate Cisplatin Insensitivity in Neuroblastoma.
Cancers
Gunda V,Pathania AS,Chava S,Prathipati P,Chaturvedi NK,Coulter DW,Pandey MK,Durden DL,Challagundla KB
Thu Sep 10 00:00:00 UTC 2020
PKCβ/NF-κB pathway in diabetic atrial remodeling.
Journal of physiology and biochemistry
Wang H,Xu Y,Xu A,Wang X,Cheng L,Lee S,Tse G,Li G,Liu T,Fu H
Sun Nov 01 00:00:00 UTC 2020
Hydrogen sulfide inhibits cigarette smoke-induced inflammation and injury in alveolar epithelial cells by suppressing PHD2/HIF-1α/MAPK signaling pathway.
International immunopharmacology
Guan R,Wang J,Li D,Li Z,Liu H,Ding M,Cai Z,Liang X,Yang Q,Long Z,Chen L,Liu W,Sun D,Yao H,Lu W
Wed Apr 01 00:00:00 UTC 2020
Exogenous CGRP Regulates Apoptosis and Autophagy to Alleviate Traumatic Brain Injury Through Akt/mTOR Signalling Pathway.
Neurochemical research
Tian J,Yang L,Wang P,Yang L,Fan Z
Tue Dec 01 00:00:00 UTC 2020
Isoquercitrin Ameliorates Cisplatin-Induced Nephrotoxicity Via the Inhibition of Apoptosis, Inflammation, and Oxidative Stress.
Frontiers in pharmacology
Wang H,Xia W,Long G,Pei Z,Li Y,Wu M,Wang Q,Zhang Y,Jia Z,Chen H
Tue Jan 12 00:00:00 UTC 2021
CAFs-derived MFAP5 promotes bladder cancer malignant behavior through NOTCH2/HEY1 signaling.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhou Z,Cui D,Sun MH,Huang JL,Deng Z,Han BM,Sun XW,Xia SJ,Sun F,Shi F
Mon Jun 01 00:00:00 UTC 2020
Integrin-α9 and Its Corresponding Ligands Play Regulatory Roles in Chronic Periodontitis.
Inflammation
Xu S,Jiang C,Liu H,Zhang H,Liao H,Wang X,Yao S,Ma L,Guo Y,Cao Z
Sat Aug 01 00:00:00 UTC 2020
Involvement of proliferative and apoptotic factors in the development of hindgut in rat fetuses with ethylenethiourea-induced anorectal malformations.
Acta histochemica
Long C,Xiao Y,Li S,Tang X,Yuan Z,Bai Y
Wed Jan 01 00:00:00 UTC 2020
Role of autophagy and apoptosis in atrophic epithelium in oral submucous fibrosis.
Journal of oral science
Zhu B,Jiang Q,Que G,Dai Z,Wu Y
Sat Mar 28 00:00:00 UTC 2020
Asiaticoside nitric oxide gel accelerates diabetic cutaneous ulcers healing by activating Wnt/β-catenin signaling pathway.
International immunopharmacology
Nie X,Zhang H,Shi X,Zhao J,Chen Y,Wu F,Yang J,Li X
Sat Feb 01 00:00:00 UTC 2020
Critical roles of microRNA-141-3p and CHD8 in hypoxia/reoxygenation-induced cardiomyocyte apoptosis.
Cell & bioscience
Yao B,Wan X,Zheng X,Zhong T,Hu J,Zhou Y,Qin A,Ma Y,Yin D
Wed Apr 13 00:00:00 UTC 2022
Notoginsenoside R1 Protects Against the Acrylamide-Induced Neurotoxicity via Upregulating Trx-1-Mediated ITGAV Expression: Involvement of Autophagy.
Frontiers in pharmacology
Wang W,Huang L,Thomas ER,Hu Y,Zeng F,Li X
Thu Oct 01 00:00:00 UTC 2020
Pseudolaric acid B induces mitotic arrest and apoptosis in both imatinib-sensitive and -resistant chronic myeloid leukaemia cells.
European journal of pharmacology
Jiang L,Wen C,He Q,Sun Y,Wang J,Lan X,Rohondia S,Dou QP,Shi X,Liu J
Fri Jun 05 00:00:00 UTC 2020
Resveratrol Downregulates STAT3 Expression and Astrocyte Activation in Primary Astrocyte Cultures of Rat.
Neurochemical research
Wu M,Wang L,Li F,Hu R,Ma J,Zhang K,Cheng X
Sat Feb 01 00:00:00 UTC 2020
Tubeimoside I improves survival of mice in sepsis by inhibiting inducible nitric oxide synthase expression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Luo M,Luo S,Cheng Z,Yang X,Lv D,Li X,Guo Y,Li C,Yan J
Mon Jun 01 00:00:00 UTC 2020
Pyrazinamide alleviates rifampin-induced steatohepatitis in mice by regulating the activities of cholesterol-activated 7α-hydroxylase and lipoprotein lipase.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Tan W,Zhao K,Xiang J,Zhou X,Cao F,Song W,Liu Q,Zhang X,Li X,Tan Z
Sat Aug 01 00:00:00 UTC 2020
MicroRNA-590-3p inhibits invasion and metastasis in triple-negative breast cancer by targeting Slug.
American journal of cancer research
Yan M,Ye L,Feng X,Shi R,Sun Z,Li Z,Liu T
Mon Sep 28 00:00:00 UTC 2020
N-Ethyl-2-Pyrrolidinone-Substituted Flavan-3-Ols with Anti-inflammatory Activity in Lipopolysaccharide-Stimulated Macrophages Are Storage-Related Marker Compounds for Green Tea.
Journal of agricultural and food chemistry
Dai W,Lou N,Xie D,Hu Z,Song H,Lu M,Shang D,Wu W,Peng J,Yin P,Lin Z
Wed Oct 28 00:00:00 UTC 2020
A multi-scale systems pharmacology approach uncovers the anti-cancer molecular mechanism of Ixabepilone.
European journal of medicinal chemistry
Cao DS,Jiang SL,Guan YD,Chen XS,Zhang LX,Zhang Y,Chen AF,Yang JM,Cheng Y
Sat Aug 01 00:00:00 UTC 2020
miR-362 knock-down promotes proliferation and inhibits apoptosis in porcine immature Sertoli cells by targeting the RMI1 gene.
Reproduction in domestic animals = Zuchthygiene
Ran M,Luo H,Gao H,Tang X,Chen Y,Zeng X,Weng B,Chen B
Fri May 01 00:00:00 UTC 2020
Cannabinoid receptor 2 activation alleviates septic lung injury by promoting autophagy via inhibition of inflammatory mediator release.
Cellular signalling
Liu AP,Yuan QH,Zhang B,Yang L,He QW,Chen K,Liu QS,Li Z,Zhan J
Fri May 01 00:00:00 UTC 2020
Clusterin contributes to hepatitis C virus-related hepatocellular carcinoma by regulating autophagy.
Life sciences
Fu N,Du H,Li D,Lu Y,Li W,Wang Y,Kong L,Du J,Zhao S,Ren W,Han F,Wang R,Zhang Y,Nan Y
Tue Sep 01 00:00:00 UTC 2020
The Establishment and Application Studies on Precise Lysosome pH Indicator Based on Self-Decomposable Nanoparticles.
Nanoscale research letters
Pang C,Song C,Li Y,Wang Q,Zhu X,Wu J,Tian Y,Fan H,Hu J,Li C,Wang B,Li X,Liu W,Fan L
Wed Jul 08 00:00:00 UTC 2020
Perilla oil regulates intestinal microbiota and alleviates insulin resistance through the PI3K/AKT signaling pathway in type-2 diabetic KKAy mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Wang J,He Y,Yu D,Jin L,Gong X,Zhang B
Wed Jan 01 00:00:00 UTC 2020
A tropomyosin-like Meretrix meretrix Linnaeus polypeptide inhibits the proliferation and metastasis of glioma cells via microtubule polymerization and FAK/Akt/MMPs signaling.
International journal of biological macromolecules
Fan Z,Xu Q,Wang C,Lin X,Zhang Q,Wu N
Sat Feb 15 00:00:00 UTC 2020
The B7-H4 gene induces immune escape partly via upregulating the PD-1/Stat3 pathway in non-small cell lung cancer.
Human immunology
Yuan L,Ye J,Fan D
Fri May 01 00:00:00 UTC 2020
Decreased Expression of the Host Long-Noncoding RNA-GM Facilitates Viral Escape by Inhibiting the Kinase activity TBK1 via S-glutathionylation.
Immunity
Wang Y,Wang P,Zhang Y,Xu J,Li Z,Li Z,Zhou Z,Liu L,Cao X
Tue Dec 15 00:00:00 UTC 2020
CCL5 derived from tumor-associated macrophages promotes prostate cancer stem cells and metastasis via activating β-catenin/STAT3 signaling.
Cell death & disease
Huang R,Wang S,Wang N,Zheng Y,Zhou J,Yang B,Wang X,Zhang J,Guo L,Wang S,Chen Z,Wang Z,Xiang S
Thu Apr 16 00:00:00 UTC 2020
A20 targets PFKL and glycolysis to inhibit the progression of hepatocellular carcinoma.
Cell death & disease
Feng Y,Zhang Y,Cai Y,Liu R,Lu M,Li T,Fu Y,Guo M,Huang H,Ou Y,Chen Y
Mon Feb 03 00:00:00 UTC 2020
TLR4 inhibition ameliorates mesencephalic substantia nigra injury in neonatal rats exposed to lipopolysaccharide via regulation of neuro-immunity.
Brain research bulletin
He W,Jiang M,Mao P,Yan F
Tue Dec 01 00:00:00 UTC 2020
The Acetone Indigo Red Dehydrating Agent IF203 Induces HepG2 Cell Death Through Cell Cycle Arrest, Autophagy and Apoptosis.
OncoTargets and therapy
Shang Y,Wang Q,Li J,Zhao Q,Huang X,Dong H,Liu H,Zhang Y,Zhang J,Gui R,Nie X
Tue Apr 12 00:00:00 UTC 2022
Colorectal cancer-derived exosomal miR-106b-3p promotes metastasis by down-regulating DLC-1 expression.
Clinical science (London, England : 1979)
Liu H,Liu Y,Sun P,Leng K,Xu Y,Mei L,Han P,Zhang B,Yao K,Li C,Bai J,Cui B
Fri Feb 28 00:00:00 UTC 2020
Antitumor activity of celastrol by inhibition of proliferation, invasion, and migration in cholangiocarcinoma via PTEN/PI3K/Akt pathway.
Cancer medicine
Zhu B,Wei Y
Wed Jan 01 00:00:00 UTC 2020
The metabolic changes in the hippocampus of an atherosclerotic rat model and the regulation of aerobic training.
Metabolic brain disease
Liu B,Li J,Lin X,Hu J,Lou S
Sat Aug 01 00:00:00 UTC 2020
Programmed cell death protein 1 promotes hepatitis B virus transmission through the regulation of ERK1/2-mediated trophoblasts differentiation.
Archives of gynecology and obstetrics
Yang J,Liu J,Sheng M,Zhang X,Liu M
Sat Feb 01 00:00:00 UTC 2020
Soluble Uric Acid Promotes Atherosclerosis via AMPK (AMP-Activated Protein Kinase)-Mediated Inflammation.
Arteriosclerosis, thrombosis, and vascular biology
Kimura Y,Yanagida T,Onda A,Tsukui D,Hosoyamada M,Kono H
Sun Mar 01 00:00:00 UTC 2020
miR-124-3p increases in high glucose induced osteocyte-derived exosomes and regulates galectin-3 expression: A possible mechanism in bone remodeling alteration in diabetic periodontitis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Guo Y,Chen YY,Liu Q,Chen Y,Tan L,Zhang SH,Gao ZR,Zhou YH,Zhang GY,Feng YZ
Sun Nov 01 00:00:00 UTC 2020
Paradoxical Pro-inflammatory Responses by Human Macrophages to an Amoebae Host-Adapted Legionella Effector.
Cell host & microbe
Price C,Jones S,Mihelcic M,Santic M,Abu Kwaik Y
Wed Apr 08 00:00:00 UTC 2020
RNA-seq analysis of potential lncRNAs for age-related hearing loss in a mouse model.
Aging
Zhao T,Liu X,Sun Z,Zhang J,Zhang X,Wang C,Geng R,Zheng T,Li B,Zheng QY
Sun Apr 26 00:00:00 UTC 2020
Guizhi Decoction () Inhibits Cholinergic Transdifferentiation by Regulating Imbalance of NGF and LIF in Salt-Sensitive Hypertensive Heart Failure Rats.
Chinese journal of integrative medicine
Wang YC,Ma DF,Jiang P,Zhang YM,Zhou GF,Yang JL,Li ZY,Li X
Sun Mar 01 00:00:00 UTC 2020
The up-regulation of NDRG1 by HIF counteracts the cancer-promoting effect of HIF in VHL-deficient clear cell renal cell carcinoma.
Cell proliferation
Zhang ZY,Zhang SL,Chen HL,Mao YQ,Li ZM,Kong CY,Han B,Zhang J,Chen YH,Xue W,Zhai W,Wang LS
Wed Jul 01 00:00:00 UTC 2020
Lamin B1 deficiency promotes malignancy and predicts poor prognosis in gastric cancer.
Neoplasma
Yu ZY,Jiang XY,Zhao RR,Luo CJ,Ren YX,Ma ZJ,Ye HL,Shi WG,Wang C,Jiao ZY
Sun Nov 01 00:00:00 UTC 2020
Protein Phosphatase 2A Inhibiting β-Catenin Phosphorylation Contributes Critically to the Anti-renal Interstitial Fibrotic Effect of Norcantharidin.
Inflammation
Xiao Z,Wen L,Zeng D,Yin D,Zhou X,Tang C,Li Y
Mon Jun 01 00:00:00 UTC 2020
Correction of Bcl-x splicing improves responses to imatinib in chronic myeloid leukaemia cells and mouse models.
British journal of haematology
Zhang J,Wang Y,Li SQ,Fang L,Wang XZ,Li J,Zhang HB,Huang B,Xu YM,Yang WM,Lin J,Min QH,Liao ZH,Wu Y,Liu J
Mon Jun 01 00:00:00 UTC 2020
LncRNA XIST promotes chemoresistance of breast cancer cells to doxorubicin by sponging miR-200c-3p to upregulate ANLN.
Clinical and experimental pharmacology & physiology
Zhang M,Wang F,Xiang Z,Huang T,Zhou WB
Sat Aug 01 00:00:00 UTC 2020
Integrated Analysis of Key Genes and Pathways Involved in Fetal Growth Restriction and Their Associations With the Dysregulation of the Maternal Immune System.
Frontiers in genetics
Wang X,Zhu H,Lei L,Zhang Y,Tang C,Wu JX,Zhou JR,Xiao XR
Tue Feb 16 00:00:00 UTC 2021
All-trans-retinal induces autophagic cell death via oxidative stress and the endoplasmic reticulum stress pathway in human retinal pigment epithelial cells.
Toxicology letters
Zhang L,Zhou Y,Xia Q,Chen Y,Li J
Wed Apr 01 00:00:00 UTC 2020
Inhibition of formyl peptide receptor 1 activity suppresses tumorigenicity in vivo and attenuates the invasion and migration of lung adenocarcinoma cells under hypoxic conditions in vitro.
Annals of translational medicine
Huang B,Guo H,Ding J,Li J,Wang H,Xu J,Zheng Q,Zhou L,Dai Q
Tue Sep 01 00:00:00 UTC 2020
Nintedanib Reduces Neutrophil Chemotaxis via Activating GRK2 in Bleomycin-Induced Pulmonary Fibrosis.
International journal of molecular sciences
Chen WC,Chen NJ,Chen HP,Yu WK,Su VY,Chen H,Wu HH,Yang KY
Thu Jul 02 00:00:00 UTC 2020
MicroRNA-150 functions as a tumor suppressor and sensitizes osteosarcoma to doxorubicin-induced apoptosis by targeting RUNX2.
Experimental and therapeutic medicine
Ling Z,Fan G,Yao D,Zhao J,Zhou Y,Feng J,Zhou G,Chen Y
Wed Jan 01 00:00:00 UTC 2020
Interferon regulatory factor 7 mediates obesity-associated MCP-1 transcription.
PloS one
Kuroda M,Nishiguchi M,Ugawa N,Ishikawa E,Kawabata Y,Okamoto S,Sasaki W,Miyatake Y,Sebe M,Masumoto S,Tsutsumi R,Harada N,Sakaue H
Wed Aug 12 00:00:00 UTC 2020
Characterization of a Novel Interleukin-1 Receptor Antagonist from Sheep (Ovis aries).
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Zhang SJ,Guo X,Hu P,Lu SY,Liu NN,Fu BQ,Wang N,Li YS,Wang LL,Chang J,Chang HZ,Liu ZS,Zhou Y,Ren HL
Fri May 01 00:00:00 UTC 2020
Thymoquinone Protects against Hyperlipidemia-Induced Cardiac Damage in Low-Density Lipoprotein Receptor-Deficient (LDL-R-/-) Mice via Its Anti-inflammatory and Antipyroptotic Effects.
BioMed research international
Pei ZW,Guo Y,Zhu HL,Dong M,Zhang Q,Wang F
Tue May 04 00:00:00 UTC 2021
WITHDRAWN: Resveratrol partially prevents learning and memory deficits in rats exposed to gaseous formaldehyde.
Environmental toxicology and pharmacology
Niu H,Xie R,Li L,Zhang X,Wei X,Deng J,Li Z
Tue Sep 22 00:00:00 UTC 2020
RIP1 promotes proliferation through G2/M checkpoint progression and mediates cisplatin-induced apoptosis and necroptosis in human ovarian cancer cells.
Acta pharmacologica Sinica
Zheng XL,Yang JJ,Wang YY,Li Q,Song YP,Su M,Li JK,Zhang L,Li ZP,Zhou B,Lin Y
Tue Sep 01 00:00:00 UTC 2020
Pharmacological Effects of Fasudil on Flap Survival in a Rodent Model.
The Journal of surgical research
Ji E,Wang J,Wang L,Pan Z,Gao W
Sun Nov 01 00:00:00 UTC 2020
Post-production modifications of murine mesenchymal stem cell (mMSC) derived extracellular vesicles (EVs) and impact on their cellular interaction.
Biomaterials
Le Saux S,Aarrass H,Lai-Kee-Him J,Bron P,Armengaud J,Miotello G,Bertrand-Michel J,Dubois E,George S,Faklaris O,Devoisselle JM,Legrand P,Chopineau J,Morille M
Sat Feb 01 00:00:00 UTC 2020
Anti-neuroinflammatory effect of 3,4-dihydroxybenzaldehyde in ischemic stroke.
International immunopharmacology
Li X,Xiang B,Shen T,Xiao C,Dai R,He F,Lin Q
Tue Mar 03 00:00:00 UTC 2020
Protection of diabetes-induced kidney injury by phosphocreatine via the regulation of ERK/Nrf2/HO-1 signaling pathway.
Life sciences
Shopit A,Niu M,Wang H,Tang Z,Li X,Tesfaldet T,Ai J,Ahmad N,Al-Azab M,Tang Z
Sat Feb 01 00:00:00 UTC 2020
Screening for reproductive biomarkers in Bactrian camel via iTRAQ analysis of proteomes.
Reproduction in domestic animals = Zuchthygiene
Wang Q,Zhang Q,Gan Z,Li H,Yang Y,Zhang Y,Zhao X
Sat Feb 01 00:00:00 UTC 2020
Ginsenoside protects against AKI via activation of HIF‑1α and VEGF‑A in the kidney‑brain axis.
International journal of molecular medicine
Mao H,Jiang C,Xu L,Chen D,Liu H,Xu Y,Ma K,Wang M
Sun Mar 01 00:00:00 UTC 2020
An ultrasensitive hybridization chain reaction-amplified CRISPR-Cas12a aptasensor for extracellular vesicle surface protein quantification.
Theranostics
Xing S,Lu Z,Huang Q,Li H,Wang Y,Lai Y,He Y,Deng M,Liu W
Mon May 24 00:00:00 UTC 2021
Involvement of p53 Acetylation in Growth Suppression of Cutaneous T-Cell Lymphomas Induced by HDAC Inhibition.
The Journal of investigative dermatology
Yu X,Li H,Zhu M,Hu P,Liu X,Qing Y,Wang X,Wang H,Wang Z,Xu J,Tan R,Guo Q,Hui H
Thu Oct 01 00:00:00 UTC 2020
Deoxycholic Acid Upregulates the Reprogramming Factors KFL4 and OCT4 Through the IL-6/STAT3 Pathway in Esophageal Adenocarcinoma Cells.
Technology in cancer research & treatment
Chen M,Ye A,Wei J,Wang R,Poon K
Mon Nov 29 00:00:00 UTC 2021
LncRNA-H19 induces hepatic stellate cell activation via upregulating alcohol dehydrogenase III-mediated retinoic acid signals.
International immunopharmacology
Wang ZM,Xia SW,Zhang T,Wang ZY,Yang X,Kai J,Cheng XD,Shao JJ,Tan SZ,Chen AP,Wang SJ,Zhang F,Zhang ZL,Zheng SZ
Wed Jul 01 00:00:00 UTC 2020
A coronary artery disease-associated tRNAThr mutation altered mitochondrial function, apoptosis and angiogenesis.
Nucleic acids research
Jia Z,Zhang Y,Li Q,Ye Z,Liu Y,Fu C,Cang X,Wang M,Guan MX
Thu Feb 28 00:00:00 UTC 2019
A coronary artery disease-associated tRNAThr mutation altered mitochondrial function, apoptosis and angiogenesis.
Nucleic acids research
Jia Z,Zhang Y,Li Q,Ye Z,Liu Y,Fu C,Cang X,Wang M,Guan MX
Thu Feb 28 00:00:00 UTC 2019
circPUM1 Promotes Tumorigenesis and Progression of Ovarian Cancer by Sponging miR-615-5p and miR-6753-5p.
Molecular therapy. Nucleic acids
Guan X,Zong ZH,Liu Y,Chen S,Wang LL,Zhao Y
Fri Dec 06 00:00:00 UTC 2019
Interferon-alpha promotes immunosuppression through IFNAR1/STAT1 signalling in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Yang W,Zhang L,Liu S,Zhao M,Zhou G,Wang L,Jin S,Zhang Z,Hu J
Fri Feb 01 00:00:00 UTC 2019
Interferon-alpha promotes immunosuppression through IFNAR1/STAT1 signalling in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Yang W,Zhang L,Liu S,Zhao M,Zhou G,Wang L,Jin S,Zhang Z,Hu J
Fri Feb 01 00:00:00 UTC 2019
CSE1L participates in regulating cell mitosis in human seminoma.
Cell proliferation
Liu C,Wei J,Xu K,Sun X,Zhang H,Xiong C
Fri Mar 01 00:00:00 UTC 2019
CSE1L participates in regulating cell mitosis in human seminoma.
Cell proliferation
Liu C,Wei J,Xu K,Sun X,Zhang H,Xiong C
Fri Mar 01 00:00:00 UTC 2019
Changes in the levels of 12/15-lipoxygenase, apoptosis-related proteins and inflammatory factors in the cortex of diabetic rats and the neuroprotection of baicalein.
Free radical biology & medicine
Li Y,Chen Q,Ran D,Wang H,Du W,Luo Y,Jiang W,Yang Y,Yang J
Mon Apr 01 00:00:00 UTC 2019
Changes in the levels of 12/15-lipoxygenase, apoptosis-related proteins and inflammatory factors in the cortex of diabetic rats and the neuroprotection of baicalein.
Free radical biology & medicine
Li Y,Chen Q,Ran D,Wang H,Du W,Luo Y,Jiang W,Yang Y,Yang J
Mon Apr 01 00:00:00 UTC 2019
Zerumbone inhibits migration in ESCC via promoting Rac1 ubiquitination.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang M,Niu J,Gao L,Gao Y,Gao S
Tue Jan 01 00:00:00 UTC 2019
Zerumbone inhibits migration in ESCC via promoting Rac1 ubiquitination.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang M,Niu J,Gao L,Gao Y,Gao S
Tue Jan 01 00:00:00 UTC 2019
FS-7 inhibits MGC-803 cells growth in vitro and in vivo via down-regulating glycolysis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Deng X,Li Z,Xiong R,Liu J,Liu R,Peng J,Chen Y,Lei X,Cao X,Zheng X,Xie Z,Tang G
Tue Jan 01 00:00:00 UTC 2019
FS-7 inhibits MGC-803 cells growth in vitro and in vivo via down-regulating glycolysis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Deng X,Li Z,Xiong R,Liu J,Liu R,Peng J,Chen Y,Lei X,Cao X,Zheng X,Xie Z,Tang G
Tue Jan 01 00:00:00 UTC 2019
The effect of di-2-ethylhexyl phthalate on inflammation and lipid metabolic disorder in rats.
Ecotoxicology and environmental safety
Zhou L,Chen H,Xu Q,Han X,Zhao Y,Song X,Zhao T,Ye L
Mon Apr 15 00:00:00 UTC 2019
The effect of di-2-ethylhexyl phthalate on inflammation and lipid metabolic disorder in rats.
Ecotoxicology and environmental safety
Zhou L,Chen H,Xu Q,Han X,Zhao Y,Song X,Zhao T,Ye L
Mon Apr 15 00:00:00 UTC 2019
Nicotinate-curcumin ameliorates cognitive impairment in diabetic rats by rescuing autophagic flux in CA1 hippocampus.
CNS neuroscience & therapeutics
Gu HF,Li N,Tang YL,Yan CQ,Shi Z,Yi SN,Zhou HL,Liao DF,OuYang XP
Mon Apr 01 00:00:00 UTC 2019
Nicotinate-curcumin ameliorates cognitive impairment in diabetic rats by rescuing autophagic flux in CA1 hippocampus.
CNS neuroscience & therapeutics
Gu HF,Li N,Tang YL,Yan CQ,Shi Z,Yi SN,Zhou HL,Liao DF,OuYang XP
Mon Apr 01 00:00:00 UTC 2019
Melatonin protects against sepsis-induced cardiac dysfunction by regulating apoptosis and autophagy via activation of SIRT1 in mice.
Life sciences
Zhang WX,He BM,Wu Y,Qiao JF,Peng ZY
Tue Jan 15 00:00:00 UTC 2019
Melatonin protects against sepsis-induced cardiac dysfunction by regulating apoptosis and autophagy via activation of SIRT1 in mice.
Life sciences
Zhang WX,He BM,Wu Y,Qiao JF,Peng ZY
Tue Jan 15 00:00:00 UTC 2019
Sulforaphane Improves Lipid Metabolism by Enhancing Mitochondrial Function and Biogenesis In Vivo and In Vitro.
Molecular nutrition & food research
Lei P,Tian S,Teng C,Huang L,Liu X,Wang J,Zhang Y,Li B,Shan Y
Fri Feb 01 00:00:00 UTC 2019
Sulforaphane Improves Lipid Metabolism by Enhancing Mitochondrial Function and Biogenesis In Vivo and In Vitro.
Molecular nutrition & food research
Lei P,Tian S,Teng C,Huang L,Liu X,Wang J,Zhang Y,Li B,Shan Y
Fri Feb 01 00:00:00 UTC 2019
DEP domain containing 1 predicts prognosis of hepatocellular carcinoma patients and regulates tumor proliferation and metastasis.
Cancer science
Qu D,Cui F,Lu D,Yang Y,Xu Y
Tue Jan 01 00:00:00 UTC 2019
DEP domain containing 1 predicts prognosis of hepatocellular carcinoma patients and regulates tumor proliferation and metastasis.
Cancer science
Qu D,Cui F,Lu D,Yang Y,Xu Y
Tue Jan 01 00:00:00 UTC 2019
Constitutive androstane receptor weakens the induction of panaxytriol on CYP3A4 by repressing the activation of pregnane X receptor.
Biochemical pharmacology
Hu Q,Yao N,Wu J,Liu M,Liu F,Zhang H,Xiong Y,Xia C
Tue Jan 01 00:00:00 UTC 2019
Constitutive androstane receptor weakens the induction of panaxytriol on CYP3A4 by repressing the activation of pregnane X receptor.
Biochemical pharmacology
Hu Q,Yao N,Wu J,Liu M,Liu F,Zhang H,Xiong Y,Xia C
Tue Jan 01 00:00:00 UTC 2019
Bromodomain inhibitor jq1 induces cell cycle arrest and apoptosis of glioma stem cells through the VEGF/PI3K/AKT signaling pathway.
International journal of oncology
Wen N,Guo B,Zheng H,Xu L,Liang H,Wang Q,Wang D,Chen X,Zhang S,Li Y,Zhang L
Tue Oct 01 00:00:00 UTC 2019
S1P promotes inflammation-induced tube formation by HLECs via the S1PR1/NF-κB pathway.
International immunopharmacology
Zheng Z,Zeng YZ,Ren K,Zhu X,Tan Y,Li Y,Li Q,Yi GH
Tue Jan 01 00:00:00 UTC 2019
S1P promotes inflammation-induced tube formation by HLECs via the S1PR1/NF-κB pathway.
International immunopharmacology
Zheng Z,Zeng YZ,Ren K,Zhu X,Tan Y,Li Y,Li Q,Yi GH
Tue Jan 01 00:00:00 UTC 2019
Hepatoprotective effects and structure-activity relationship of five flavonoids against lipopolysaccharide/d-galactosamine induced acute liver failure in mice.
International immunopharmacology
He Y,Xia Z,Yu D,Wang J,Jin L,Huang D,Ye X,Li X,Zhang B
Fri Mar 01 00:00:00 UTC 2019
Hepatoprotective effects and structure-activity relationship of five flavonoids against lipopolysaccharide/d-galactosamine induced acute liver failure in mice.
International immunopharmacology
He Y,Xia Z,Yu D,Wang J,Jin L,Huang D,Ye X,Li X,Zhang B
Fri Mar 01 00:00:00 UTC 2019
Overexpression of CDCA8 promotes the malignant progression of cutaneous melanoma and leads to poor prognosis.
International journal of molecular medicine
Ci C,Tang B,Lyu D,Liu W,Qiang D,Ji X,Qiu X,Chen L,Ding W
Tue Jan 01 00:00:00 UTC 2019
Overexpression of CDCA8 promotes the malignant progression of cutaneous melanoma and leads to poor prognosis.
International journal of molecular medicine
Ci C,Tang B,Lyu D,Liu W,Qiang D,Ji X,Qiu X,Chen L,Ding W
Tue Jan 01 00:00:00 UTC 2019
Radioprotective effects of roxadustat (FG-4592) in haematopoietic system.
Journal of cellular and molecular medicine
Zhang P,Du J,Zhao H,Cheng Y,Dong S,Yang Y,Li B,Gao F,Sun X,Cai J,Liu C
Tue Jan 01 00:00:00 UTC 2019
Radioprotective effects of roxadustat (FG-4592) in haematopoietic system.
Journal of cellular and molecular medicine
Zhang P,Du J,Zhao H,Cheng Y,Dong S,Yang Y,Li B,Gao F,Sun X,Cai J,Liu C
Tue Jan 01 00:00:00 UTC 2019
miR-429 and miR-424-5p inhibit cell proliferation and Ca2+ influx by downregulating CaSR in pulmonary artery smooth muscle cells.
American journal of physiology. Cell physiology
Li C,Qin F,Xue M,Lei Y,Hu F,Xu H,Sun G,Wang T,Guo M
Tue Jan 01 00:00:00 UTC 2019
miR-429 and miR-424-5p inhibit cell proliferation and Ca2+ influx by downregulating CaSR in pulmonary artery smooth muscle cells.
American journal of physiology. Cell physiology
Li C,Qin F,Xue M,Lei Y,Hu F,Xu H,Sun G,Wang T,Guo M
Tue Jan 01 00:00:00 UTC 2019
iTRAQ-based quantitative proteomics reveals the neuroprotection of rhubarb in experimental intracerebral hemorrhage.
Journal of ethnopharmacology
Liu T,Zhou J,Cui H,Li P,Luo J,Li T,He F,Wang Y,Tang T
Mon Mar 25 00:00:00 UTC 2019
iTRAQ-based quantitative proteomics reveals the neuroprotection of rhubarb in experimental intracerebral hemorrhage.
Journal of ethnopharmacology
Liu T,Zhou J,Cui H,Li P,Luo J,Li T,He F,Wang Y,Tang T
Mon Mar 25 00:00:00 UTC 2019
S-1-Propenylcysteine augments BACH1 degradation and heme oxygenase 1 expression in a nitric oxide-dependent manner in endothelial cells.
Nitric oxide : biology and chemistry
Tsuneyoshi T,Kunimura K,Morihara N
Fri Mar 01 00:00:00 UTC 2019
S-1-Propenylcysteine augments BACH1 degradation and heme oxygenase 1 expression in a nitric oxide-dependent manner in endothelial cells.
Nitric oxide : biology and chemistry
Tsuneyoshi T,Kunimura K,Morihara N
Fri Mar 01 00:00:00 UTC 2019
Hypericum perforatum L. supplementation protects sciatic nerve injury-induced apoptotic, inflammatory and oxidative damage to muscle, blood and brain in rats.
The Journal of pharmacy and pharmacology
Uslusoy F,Nazıroğlu M,Övey İS,Sönmez TT
Tue Jan 01 00:00:00 UTC 2019
Hypericum perforatum L. supplementation protects sciatic nerve injury-induced apoptotic, inflammatory and oxidative damage to muscle, blood and brain in rats.
The Journal of pharmacy and pharmacology
Uslusoy F,Nazıroğlu M,Övey İS,Sönmez TT
Tue Jan 01 00:00:00 UTC 2019
Prolonged high-glucose exposure decreased SREBP-1/FASN/ACC in Schwann cells of diabetic mice via blocking PI3K/Akt pathway.
Journal of cellular biochemistry
Zhu L,Du W,Liu Y,Cheng M,Wang X,Zhang C,Lv X,Li F,Zhao S,Hao J
Mon Apr 01 00:00:00 UTC 2019
Prolonged high-glucose exposure decreased SREBP-1/FASN/ACC in Schwann cells of diabetic mice via blocking PI3K/Akt pathway.
Journal of cellular biochemistry
Zhu L,Du W,Liu Y,Cheng M,Wang X,Zhang C,Lv X,Li F,Zhao S,Hao J
Mon Apr 01 00:00:00 UTC 2019
GDF-15 promotes mitochondrial function and proliferation in neuronal HT22 cells.
Journal of cellular biochemistry
Liu H,Liu J,Si L,Guo C,Liu W,Liu Y
Sat Jun 01 00:00:00 UTC 2019
GDF-15 promotes mitochondrial function and proliferation in neuronal HT22 cells.
Journal of cellular biochemistry
Liu H,Liu J,Si L,Guo C,Liu W,Liu Y
Sat Jun 01 00:00:00 UTC 2019
Up-regulation of EMT-related gene VCAN by NPM1 mutant-driven TGF-β/cPML signalling promotes leukemia cell invasion.
Journal of Cancer
Yang L,Wang L,Yang Z,Jin H,Zou Q,Zhan Q,Tang Y,Tao Y,Lei L,Jing Y,Jiang X,Zhang L
Thu Oct 01 00:00:00 UTC 2020
Differential response to lead toxicity in rat primary microglia and astrocytes.
Toxicology and applied pharmacology
Peng J,Zhou F,Wang Y,Xu Y,Zhang H,Zou F,Meng X
Tue Jan 15 00:00:00 UTC 2019
Differential response to lead toxicity in rat primary microglia and astrocytes.
Toxicology and applied pharmacology
Peng J,Zhou F,Wang Y,Xu Y,Zhang H,Zou F,Meng X
Tue Jan 15 00:00:00 UTC 2019
Regulation of Neurotransmitter Release by Amyloid Precursor Protein Through Synapsin Phosphorylation.
Neurochemical research
Liu A,Zhang Y,Han L,He G,Xie W,Zhou Z,Jia Z
Fri Mar 01 00:00:00 UTC 2019
Regulation of Neurotransmitter Release by Amyloid Precursor Protein Through Synapsin Phosphorylation.
Neurochemical research
Liu A,Zhang Y,Han L,He G,Xie W,Zhou Z,Jia Z
Fri Mar 01 00:00:00 UTC 2019
High-mobility group box 1 protein modulated proliferation and radioresistance in esophageal squamous cell carcinoma.
Journal of gastroenterology and hepatology
Di X,He G,Chen H,Zhu C,Qin Q,Yan J,Zhang X,Sun X
Mon Apr 01 00:00:00 UTC 2019
High-mobility group box 1 protein modulated proliferation and radioresistance in esophageal squamous cell carcinoma.
Journal of gastroenterology and hepatology
Di X,He G,Chen H,Zhu C,Qin Q,Yan J,Zhang X,Sun X
Mon Apr 01 00:00:00 UTC 2019
Regulation of host cell pyroptosis and cytokines production by Mycobacterium tuberculosis effector PPE60 requires LUBAC mediated NF-κB signaling.
Cellular immunology
Gong Z,Kuang Z,Li H,Li C,Ali MK,Huang F,Li P,Li Q,Huang X,Ren S,Li J,Xie J
Tue Jan 01 00:00:00 UTC 2019
Regulation of host cell pyroptosis and cytokines production by Mycobacterium tuberculosis effector PPE60 requires LUBAC mediated NF-κB signaling.
Cellular immunology
Gong Z,Kuang Z,Li H,Li C,Ali MK,Huang F,Li P,Li Q,Huang X,Ren S,Li J,Xie J
Tue Jan 01 00:00:00 UTC 2019
Mechanism Underlying the Protective Effect of Selenium on NO-Induced Oxidative Damage in Bovine Mammary Epithelial Cells.
Biological trace element research
Guo Y,Guo X,Yan S,Zhang B,Shi B
Sun Sep 01 00:00:00 UTC 2019
Long noncoding RNA HOTAIR regulates autophagy via the miR-20b-5p/ATG7 axis in hepatic ischemia/reperfusion injury.
Gene
Tang B,Bao N,He G,Wang J
Wed Feb 20 00:00:00 UTC 2019
Long noncoding RNA HOTAIR regulates autophagy via the miR-20b-5p/ATG7 axis in hepatic ischemia/reperfusion injury.
Gene
Tang B,Bao N,He G,Wang J
Wed Feb 20 00:00:00 UTC 2019
SOCS3 control the activity of NF-κB induced by HSP70 via degradation of MyD88-adapter-like protein (Mal) in IPEC-J2 cells.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group
Yong YH,Wang P,Jia RM,Gooneratne R,Robert Wang HC,Liao M,Ju XH
Mon Jan 06 00:00:00 UTC 2020
SOCS3 control the activity of NF-κB induced by HSP70 via degradation of MyD88-adapter-like protein (Mal) in IPEC-J2 cells.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group
Yong YH,Wang P,Jia RM,Gooneratne R,Robert Wang HC,Liao M,Ju XH
Mon Jan 06 00:00:00 UTC 2020
Repurposing of omeprazole for oligodendrocyte differentiation and remyelination.
Brain research
Zhu K,Sun J,Kang Z,Zou Z,Wu X,Wang Y,Wu G,Harris RA,Wang J
Wed May 01 00:00:00 UTC 2019
Repurposing of omeprazole for oligodendrocyte differentiation and remyelination.
Brain research
Zhu K,Sun J,Kang Z,Zou Z,Wu X,Wang Y,Wu G,Harris RA,Wang J
Wed May 01 00:00:00 UTC 2019
MARCH5 overexpression contributes to tumor growth and metastasis and associates with poor survival in breast cancer.
Cancer management and research
Tang H,Peng S,Dong Y,Yang X,Yang P,Yang L,Yang B,Bao G
Thu Mar 31 00:00:00 UTC 2022
MARCH5 overexpression contributes to tumor growth and metastasis and associates with poor survival in breast cancer.
Cancer management and research
Tang H,Peng S,Dong Y,Yang X,Yang P,Yang L,Yang B,Bao G
Thu Mar 31 00:00:00 UTC 2022
PARP inhibitor re‑sensitizes Adriamycin resistant leukemia cells through DNA damage and apoptosis.
Molecular medicine reports
Wu J,Xiao S,Yuan M,Li Q,Xiao G,Wu W,Ouyang Y,Huang L,Yao C
Tue Jan 01 00:00:00 UTC 2019
PARP inhibitor re‑sensitizes Adriamycin resistant leukemia cells through DNA damage and apoptosis.
Molecular medicine reports
Wu J,Xiao S,Yuan M,Li Q,Xiao G,Wu W,Ouyang Y,Huang L,Yao C
Tue Jan 01 00:00:00 UTC 2019
Glucose metabolic alterations in hippocampus of diabetes mellitus rats and the regulation of aerobic exercise.
Behavioural brain research
Li J,Liu B,Cai M,Lin X,Lou S
Fri May 17 00:00:00 UTC 2019
Glucose metabolic alterations in hippocampus of diabetes mellitus rats and the regulation of aerobic exercise.
Behavioural brain research
Li J,Liu B,Cai M,Lin X,Lou S
Fri May 17 00:00:00 UTC 2019
Simvastatin ameliorates total liver ischemia/reperfusion injury via KLF2-mediated mechanism in rats.
Clinics and research in hepatology and gastroenterology
Liu Z,Lai CH,Zhang X,Luo J,Huang X,Qi X,Wang W,Zhong Z,Xiaoli F,Li L,Xiong Y,Senninger N,Wang Y,Ye Q,Ye S
Mon Apr 01 00:00:00 UTC 2019
Simvastatin ameliorates total liver ischemia/reperfusion injury via KLF2-mediated mechanism in rats.
Clinics and research in hepatology and gastroenterology
Liu Z,Lai CH,Zhang X,Luo J,Huang X,Qi X,Wang W,Zhong Z,Xiaoli F,Li L,Xiong Y,Senninger N,Wang Y,Ye Q,Ye S
Mon Apr 01 00:00:00 UTC 2019
HepaCAM Regulates Warburg Effect of Renal Cell Carcinoma via HIF-1α/NF-κB Signaling Pathway.
Urology
Fan Y,Ou L,Fan J,Li L,Wu X,Luo C,Gao Y,Niu L
Wed May 01 00:00:00 UTC 2019
HepaCAM Regulates Warburg Effect of Renal Cell Carcinoma via HIF-1α/NF-κB Signaling Pathway.
Urology
Fan Y,Ou L,Fan J,Li L,Wu X,Luo C,Gao Y,Niu L
Wed May 01 00:00:00 UTC 2019
Exogenous Tetranectin Protects Against 1-Methyl-4-Phenylpyridine-Induced Neurotoxicity by Inhibiting Apoptosis and Autophagy Through Ribosomal Protein S6 Kinase Beta-1.
World neurosurgery
Xie Q,Liu M,Yan YF,Shen X,Wang ES
Fri Feb 01 00:00:00 UTC 2019
Exogenous Tetranectin Protects Against 1-Methyl-4-Phenylpyridine-Induced Neurotoxicity by Inhibiting Apoptosis and Autophagy Through Ribosomal Protein S6 Kinase Beta-1.
World neurosurgery
Xie Q,Liu M,Yan YF,Shen X,Wang ES
Fri Feb 01 00:00:00 UTC 2019
Circ-ASH2L promotes tumor progression by sponging miR-34a to regulate Notch1 in pancreatic ductal adenocarcinoma.
Journal of experimental & clinical cancer research : CR
Chen Y,Li Z,Zhang M,Wang B,Ye J,Zhang Y,Tang D,Ma D,Jin W,Li X,Wang S
Tue Nov 12 00:00:00 UTC 2019
Design and Synthesis of Potent, Selective Inhibitors of Protein Arginine Methyltransferase 4 against Acute Myeloid Leukemia.
Journal of medicinal chemistry
Guo Z,Zhang Z,Yang H,Cao D,Xu X,Zheng X,Chen D,Wang Q,Li Y,Li J,Du Z,Wang X,Chen L,Ding J,Shen J,Geng M,Huang X,Xiong B
Thu Jun 13 00:00:00 UTC 2019
Hypertension-associated mitochondrial DNA 4401A>G mutation caused the aberrant processing of tRNAMet, all 8 tRNAs and ND6 mRNA in the light-strand transcript.
Nucleic acids research
Zhao X,Cui L,Xiao Y,Mao Q,Aishanjiang M,Kong W,Liu Y,Chen H,Hong F,Jia Z,Wang M,Jiang P,Guan MX
Mon Nov 04 00:00:00 UTC 2019
Microglial LOX-1/MAPKs/NF-κB positive loop promotes the vicious cycle of neuroinflammation and neural injury.
International immunopharmacology
Ge X,Zhang DM,Li MM,Zhang Y,Zhu XY,Zhou Y,Peng X,Shen AG
Wed May 01 00:00:00 UTC 2019
Babao Dan induces gastric cancer cell apoptosis via regulating MAPK and NF-κB signaling pathways.
The Journal of international medical research
Shang H,Cao Z,Zhao J,Guan J,Liu J,Peng J,Chen Y,Joseph Sferra T,Sankararaman S,Lin J
Tue Oct 01 00:00:00 UTC 2019
REG4 is an indicator for KRAS mutant lung adenocarcinoma with TTF-1 low expression.
Journal of cancer research and clinical oncology
Sun S,Hu Z,Huang S,Ye X,Wang J,Chang J,Wu X,Wang Q,Zhang L,Hu X,Yu H
Sun Sep 01 00:00:00 UTC 2019
The Anticonvulsant Effects of Baldrinal on Pilocarpine-Induced convulsion in Adult Male Mice.
Molecules (Basel, Switzerland)
Zhang X,Li X,Liu N,Zheng P,Ma L,Guo F,Sun T,Zhou R,Yu J
Wed Apr 24 00:00:00 UTC 2019
Predictable chronic mild stress promotes recovery from LPS-induced depression.
Molecular brain
Dang R,Guo YY,Zhang K,Jiang P,Zhao MG
Fri May 03 00:00:00 UTC 2019
Wnt7a predicts poor prognosis, and contributes to growth and metastasis in tongue squamous cell carcinoma.
Oncology reports
Jia B,Qiu X,Chu H,Sun X,Xu S,Zhao X,Zhao J
Fri Mar 01 00:00:00 UTC 2019
Autophagy inhibition potentiates the anti-angiogenic property of multikinase inhibitor anlotinib through JAK2/STAT3/VEGFA signaling in non-small cell lung cancer cells.
Journal of experimental & clinical cancer research : CR
Liang L,Hui K,Hu C,Wen Y,Yang S,Zhu P,Wang L,Xia Y,Qiao Y,Sun W,Fei J,Chen T,Zhao F,Yang B,Jiang X
Tue Feb 12 00:00:00 UTC 2019
Bexarotene Attenuates Focal Cerebral Ischemia-Reperfusion Injury via the Suppression of JNK/Caspase-3 Signaling Pathway.
Neurochemical research
Liu H,Liu S,Tian X,Wang Q,Rao J,Wang Y,Xiang F,Zheng H,Xu L,Dong Z
Sun Dec 01 00:00:00 UTC 2019
PP2Ac Modulates AMPK-Mediated Induction of Autophagy in Mycobacterium bovis-Infected Macrophages.
International journal of molecular sciences
Hussain T,Zhao D,Shah SZA,Sabir N,Wang J,Liao Y,Song Y,Hussain Mangi M,Yao J,Dong H,Yang L,Zhou X
Fri Nov 29 00:00:00 UTC 2019
miR-205-5p regulates epithelial-mesenchymal transition by targeting PTEN via PI3K/AKT signaling pathway in cisplatin-resistant nasopharyngeal carcinoma cells.
Gene
Zhang P,Lu X,Shi Z,Li X,Zhang Y,Zhao S,Liu H
Tue Aug 20 00:00:00 UTC 2019
Protective effects of Clec11a in islets against lipotoxicity via modulation of proliferation and lipid metabolism in mice.
Experimental cell research
Shi R,Hu J,Li W,Wang Z,Pan Y,Bai M,Mao W,Wang X,Zhong M,Yuan Y,Lau J,Sun Z,Zhao S
Fri Nov 01 00:00:00 UTC 2019
Dehydroepiandrosterone alleviates E. Coli O157:H7-induced inflammation by preventing the activation of p38 MAPK and NF-κB pathways in mice peritoneal macrophages.
Molecular immunology
Zhao J,Cao J,Yu L,Ma H
Tue Oct 01 00:00:00 UTC 2019
Glycogen phosphorylase B promotes ovarian cancer progression via Wnt/β-catenin signaling and is regulated by miR-133a-3p.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhou Y,Jin Z,Wang C
Sun Dec 01 00:00:00 UTC 2019
Role of PM2.5 in the development and progression of COPD and its mechanisms.
Respiratory research
Zhao J,Li M,Wang Z,Chen J,Zhao J,Xu Y,Wei X,Wang J,Xie J
Thu Jun 13 00:00:00 UTC 2019
Aloperine suppresses human pulmonary vascular smooth muscle cell proliferation via inhibiting inflammatory response.
The Chinese journal of physiology
Chang Z,Zhang P,Zhang M,Jun F,Hu Z,Yang J,Wu Y,Zhou R
Tue Oct 01 00:00:00 UTC 2019
Strand break-induced replication fork collapse leads to C-circles, C-overhangs and telomeric recombination.
PLoS genetics
Zhang T,Zhang Z,Shengzhao G,Li X,Liu H,Zhao Y
Fri Feb 01 00:00:00 UTC 2019
Carnosine Protects Mouse Podocytes from High Glucose Induced Apoptosis through PI3K/AKT and Nrf2 Pathways.
BioMed research international
Zhao K,Li Y,Wang Z,Han N,Wang Y
Thu Dec 12 00:00:00 UTC 2019
STAT3 Contributes To Oncolytic Newcastle Disease Virus-Induced Immunogenic Cell Death in Melanoma Cells.
Frontiers in oncology
Shao X,Wang X,Guo X,Jiang K,Ye T,Chen J,Fang J,Gu L,Wang S,Zhang G,Meng S,Xu Q
Wed Sep 30 00:00:00 UTC 2020
Oncogenic ADAM28 induces gemcitabine resistance and predicts a poor prognosis in pancreatic cancer.
World journal of gastroenterology
Wei L,Wen JY,Chen J,Ma XK,Wu DH,Chen ZH,Huang JL
Mon Oct 07 00:00:00 UTC 2019
LncRNA-MTA2TR functions as a promoter in pancreatic cancer via driving deacetylation-dependent accumulation of HIF-1α.
Theranostics
Zeng Z,Xu FY,Zheng H,Cheng P,Chen QY,Ye Z,Zhong JX,Deng SJ,Liu ML,Huang K,Li Q,Li W,Hu YH,Wang F,Wang CY,Zhao G
Thu Aug 20 00:00:00 UTC 2020
CircRhoC promotes tumorigenicity and progression in ovarian cancer by functioning as a miR-302e sponge to positively regulate VEGFA.
Journal of cellular and molecular medicine
Wang LL,Zong ZH,Liu Y,Guan X,Chen S,Zhao Y
Sun Dec 01 00:00:00 UTC 2019
Nrf2/Keap1/ARE Signaling Mediated an Antioxidative Protection of Human Placental Mesenchymal Stem Cells of Fetal Origin in Alveolar Epithelial Cells.
Oxidative medicine and cellular longevity
Yan X,Fu X,Jia Y,Ma X,Tao J,Yang T,Ma H,Liang X,Liu X,Yang J,Wei J
Fri Dec 13 00:00:00 UTC 2019
The PERK-EIF2α-ATF4 signaling branch regulates osteoblast differentiation and proliferation by PTH.
American journal of physiology. Endocrinology and metabolism
Zhang K,Wang M,Li Y,Li C,Tang S,Qu X,Feng N,Wu Y
Mon Apr 01 00:00:00 UTC 2019
Prognostic implications and oncogenic roles of MYBL2 protein expression in esophageal squamous-cell carcinoma.
OncoTargets and therapy
Qin H,Li Y,Zhang H,Wang F,He H,Bai X,Li S
Fri Apr 08 00:00:00 UTC 2022
The up-regulation of two identified wound healing specific proteins-HSP70 and lysozyme in regenerated Eisenia fetida through transcriptome analysis.
Journal of ethnopharmacology
Yang Y,Sun Y,Zhang N,Li J,Zhang C,Duan X,Ding Y,Zhao R,Zheng Z,Geng D,Sun Y
Wed Jun 12 00:00:00 UTC 2019
TGF-β1 Induces Epithelial-Mesenchymal Transition of Chronic Sinusitis with Nasal Polyps through MicroRNA-21.
International archives of allergy and immunology
Li X,Li C,Zhu G,Yuan W,Xiao ZA
Thu Sep 12 00:00:00 UTC 2019
Spica Prunellae Extract Enhances Fluorouracil Sensitivity of 5-Fluorouracil-Resistant Human Colon Carcinoma HCT-8/5-FU Cells via TOP2α and miR-494.
BioMed research international
Fang Y,Yang C,Zhang L,Wei L,Lin J,Zhao J,Peng J
Thu Mar 19 00:00:00 UTC 2020
TRIM28 protects CARM1 from proteasome-mediated degradation to prevent colorectal cancer metastasis.
Science bulletin
Cui J,Hu J,Ye Z,Fan Y,Li Y,Wang G,Wang L,Wang Z
Tue Jul 30 00:00:00 UTC 2019
miR-629-5p promotes growth and metastasis of hepatocellular carcinoma by activating β-catenin.
Experimental cell research
Tao X,Yang X,Wu K,Yang L,Huang Y,Jin Q,Chen S
Mon Jul 15 00:00:00 UTC 2019
Cadmium regulates von Willebrand factor and occludin expression in glomerular endothelial cells of mice in a TNF-α-dependent manner.
Renal failure
Sun Z,Xie Q,Pan J,Niu N
Fri Nov 01 00:00:00 UTC 2019
CITED1 contributes to the progression of papillary thyroid carcinoma via the Wnt/β-catenin signaling pathway.
OncoTargets and therapy
Wang Y,Huang H,Hu F,Li J,Zhang L,Pang H
Mon Apr 11 00:00:00 UTC 2022
HMGB1 contributes to SASH1 methylation to attenuate astrocyte adhesion.
Cell death & disease
Wu R,Yan Y,Ma C,Chen H,Dong Z,Wang Y,Liu Y,Liu M,Yang L
Tue May 28 00:00:00 UTC 2019
TCF7L1 indicates prognosis and promotes proliferation through activation of Keap1/NRF2 in gastric cancer.
Acta biochimica et biophysica Sinica
Zhang B,Wu J,Cai Y,Luo M,Wang B,Gu Y
Mon Apr 01 00:00:00 UTC 2019
Nano-Gold Loaded with Resveratrol Enhance the Anti-Hepatoma Effect of Resveratrol In Vitro and In Vivo.
Journal of biomedical nanotechnology
Zhang D,Zhang J,Zeng J,Li Z,Zuo H,Huang C,Zhao X
Fri Feb 01 00:00:00 UTC 2019
Cordycepin alleviates hepatic lipid accumulation by inducing protective autophagy via PKA/mTOR pathway.
Biochemical and biophysical research communications
Li T,Wen L,Cheng B
Tue Aug 27 00:00:00 UTC 2019
MiR-146b accelerates osteoarthritis progression by targeting alpha-2-macroglobulin.
Aging
Liu X,Liu L,Zhang H,Shao Y,Chen Z,Feng X,Fang H,Zhao C,Pan J,Zhang H,Zeng C,Cai D
Sat Aug 17 00:00:00 UTC 2019
Sericin Nerve Guidance Conduit Delivering Therapeutically Repurposed Clobetasol for Functional and Structural Regeneration of Transected Peripheral Nerves.
ACS biomaterials science & engineering
Zhang L,Yang W,Xie H,Wang H,Wang J,Su Q,Li X,Song Y,Wang G,Wang L,Wang Z
Mon Mar 11 00:00:00 UTC 2019
Citrate reduced oxidative damage in stem cells by regulating cellular redox signaling pathways and represent a potential treatment for oxidative stress-induced diseases.
Redox biology
Wu X,Dai H,Liu L,Xu C,Yin Y,Yi J,Bielec MD,Han Y,Li S
Fri Feb 01 00:00:00 UTC 2019
Citrate reduced oxidative damage in stem cells by regulating cellular redox signaling pathways and represent a potential treatment for oxidative stress-induced diseases.
Redox biology
Wu X,Dai H,Liu L,Xu C,Yin Y,Yi J,Bielec MD,Han Y,Li S
Fri Feb 01 00:00:00 UTC 2019
Effects of myostatin deficiency on PGC-1α and FNDC5 expression in three different murine muscle types.
Acta histochemica
Bai Y,Bi H,Li L,Li J,Yu X,Ren H,Li Y,Ji Y,Li H,Wang H
Mon Apr 01 00:00:00 UTC 2019
OPA1 overexpression ameliorates mitochondrial cristae remodeling, mitochondrial dysfunction, and neuronal apoptosis in prion diseases.
Cell death & disease
Wu W,Zhao D,Shah SZA,Zhang X,Lai M,Yang D,Wu X,Guan Z,Li J,Zhao H,Li W,Gao H,Zhou X,Qiao J,Yang L
Tue Sep 24 00:00:00 UTC 2019
The Wnt/β-Catenin/LEF1 Pathway Promotes Cell Proliferation at Least in Part Through Direct Upregulation of miR-17-92 Cluster.
Frontiers in genetics
Mu F,Huang J,Xing T,Jing Y,Cui T,Guo Y,Yan X,Li H,Wang N
Wed Sep 30 00:00:00 UTC 2020
Propofol attenuates TNF-α-induced MMP-9 expression in human cerebral microvascular endothelial cells by inhibiting Ca2+/CAMK II/ERK/NF-κB signaling pathway.
Acta pharmacologica Sinica
Ding XW,Sun X,Shen XF,Lu Y,Wang JQ,Sun ZR,Miao CH,Chen JW
Tue Oct 01 00:00:00 UTC 2019
The IRE1α-XBP1 pathway function in hypoxia-induced pulmonary vascular remodeling, is upregulated by quercetin, inhibits apoptosis and partially reverses the effect of quercetin in PASMCs.
American journal of translational research
Cao X,He Y,Li X,Xu Y,Liu X
Thu Oct 01 00:00:00 UTC 2020
Precise and systematic survey of the efficacy of multicomponent drugs against functional dyspepsia.
Scientific reports
Wei J,Man Q,Guo F,Xian M,Wang T,Tang C,Zhang Y,Li D,Tang D,Yang H,Huang L
Wed Jul 24 00:00:00 UTC 2019
Atorvastatin Inhibits Breast Cancer Cells by Downregulating PTEN/AKT Pathway via Promoting Ras Homolog Family Member B (RhoB).
BioMed research international
Ma Q,Gao Y,Xu P,Li K,Xu X,Gao J,Qi Y,Xu J,Yang Y,Song W,He X,Liu S,Yuan X,Yin W,He Y,Pan W,Wei L,Zhang J
Thu Aug 15 00:00:00 UTC 2019
Resveratrol inhibits the invasion and metastasis of colon cancer through reversal of epithelial‑ mesenchymal transition via the AKT/GSK‑3β/Snail signaling pathway.
Molecular medicine reports
Yuan L,Zhou M,Huang D,Wasan HS,Zhang K,Sun L,Huang H,Ma S,Shen M,Ruan S
Sun Sep 01 00:00:00 UTC 2019
LPS Inhibits Fatty Acid Absorption in Enterocytes through TNF-α Secreted by Macrophages.
Cells
Liu H,Kai L,Du H,Wang X,Wang Y
Thu Dec 12 00:00:00 UTC 2019
Protective effects of 18β-glycyrrhetinic acid on pulmonary arterial hypertension via regulation of Rho A/Rho kinsase pathway.
Chemico-biological interactions
Zhang M,Chang Z,Zhang P,Jing Z,Yan L,Feng J,Hu Z,Xu Q,Zhou W,Ma P,Hao Y,Zhou R
Wed Sep 25 00:00:00 UTC 2019
Wnt1 silencing enhances neurotoxicity induced by paraquat and maneb in SH-SY5Y cells.
Experimental and therapeutic medicine
Huang C,Ma J,Li BX,Sun Y
Fri Nov 01 00:00:00 UTC 2019
Nek2 augments sorafenib resistance by regulating the ubiquitination and localization of β-catenin in hepatocellular carcinoma.
Journal of experimental & clinical cancer research : CR
Deng L,Sun J,Chen X,Liu L,Wu D
Thu Jul 18 00:00:00 UTC 2019
Acid sphingomyelinase mediates the noise-induced liver disorder in mice.
Clinical and experimental pharmacology & physiology
Meng X,Gu Z,Xie X,Su Y,Zhang X,Ma H,Guo Y,Liu X,Cheng Y,Chang Y,Bao J
Sat Jun 01 00:00:00 UTC 2019
MiRNA-29a modulates visceral hyperalgesia in irritable bowel syndrome by targeting HTR7.
Biochemical and biophysical research communications
Zhu H,Xiao X,Chai Y,Li D,Yan X,Tang H
Tue Apr 09 00:00:00 UTC 2019
AAV-Mig-6 Increase the Efficacy of TAE in VX2 Rabbit Model, Is Associated With JNK Mediated Autophagy.
Journal of Cancer
Li Z,Tian Y,Qu L,Mao J,Zhong H
Thu Oct 01 00:00:00 UTC 2020
ROS-mediated inactivation of the PI3K/AKT pathway is involved in the antigastric cancer effects of thioredoxin reductase-1 inhibitor chaetocin.
Cell death & disease
Wen C,Wang H,Wu X,He L,Zhou Q,Wang F,Chen S,Huang L,Chen J,Wang H,Ye W,Li W,Yang X,Liu H,Peng J
Thu Oct 24 00:00:00 UTC 2019
miR-181b-5p May Regulate Muscle Growth in Tilapia by Targeting Myostatin b.
Frontiers in endocrinology
Zhao Z,Yu X,Jia J,Yang G,Sun C,Li W
Fri Dec 20 00:00:00 UTC 2019
Reduction of SIRT1 blunts the protective effects of ischemic post-conditioning in diabetic mice by impairing the Akt signaling pathway.
Biochimica et biophysica acta. Molecular basis of disease
Ding M,Hu L,Yang H,Gao C,Zeng K,Yu M,Feng J,Qiu J,Liu C,Fu F,Li Y
Sat Jun 01 00:00:00 UTC 2019
Toxicity mechanism of sevoflurane in neural stem cells of rats through DNA methylation.
Experimental and therapeutic medicine
Wang K,Tian Y,Zhang Y,Li X,Wei X,Hu H,Xu S
Mon Jul 01 00:00:00 UTC 2019
Novel diterpenoid-type activators of the Keap1/Nrf2/ARE signaling pathway and their regulation of redox homeostasis.
Free radical biology & medicine
Li AL,Shen T,Wang T,Zhou MX,Wang B,Song JT,Zhang PL,Wang XL,Ren DM,Lou HX,Wang XN
Sun Sep 01 00:00:00 UTC 2019
LncRNA ZFAS1 promotes proliferation and migration and inhibits apoptosis in nasopharyngeal carcinoma via the PI3K/AKT pathway in vitro.
Cancer biomarkers : section A of Disease markers
Wang X,Jin Q,Wang X,Chen W,Cai Z
Fri Mar 20 00:00:00 UTC 2020
Echinacoside Alleviates Hypoxic-Ischemic Brain Injury in Neonatal Rat by Enhancing Antioxidant Capacity and Inhibiting Apoptosis.
Neurochemical research
Wei W,Lan XB,Liu N,Yang JM,Du J,Ma L,Zhang WJ,Niu JG,Sun T,Yu JQ
Mon Jul 01 00:00:00 UTC 2019
Ginsenoside Rk1 induces cell cycle arrest and apoptosis in MDA-MB-231 triple negative breast cancer cells.
Toxicology
Hong Y,Fan D
Mon Apr 15 00:00:00 UTC 2019
Disubstituted Dithiolethione ACDT Exerts Neuroprotective Effects Against 6-Hydroxydopamine-Induced Oxidative Stress in SH-SY5Y Cells.
Neurochemical research
Betharia S,Rondόn-Ortiz AN,Brown DA
Thu Aug 01 00:00:00 UTC 2019
Blockade of glycolysis-dependent contraction by oroxylin a via inhibition of lactate dehydrogenase-a in hepatic stellate cells.
Cell communication and signaling : CCS
Wang F,Jia Y,Li M,Wang L,Shao J,Guo Q,Tan S,Ding H,Chen A,Zhang F,Zheng S
Mon Feb 11 00:00:00 UTC 2019
The long non-coding RNA H19 induces hypoxia/reoxygenation injury by up-regulating autophagy in the hepatoma carcinoma cells.
Biological research
Cui C,Li Z,Wu D
Thu Jun 13 00:00:00 UTC 2019
LncRNA SLCO4A1-AS1 predicts poor prognosis and promotes proliferation and metastasis via the EGFR/MAPK pathway in colorectal cancer.
International journal of biological sciences
Tang R,Chen J,Tang M,Liao Z,Zhou L,Jiang J,Hu Y,Liao Q,Xiong W,Tang Y,Nie S
Mon Jun 01 00:00:00 UTC 2020
c‑Jun N‑terminal kinase/transforming growth factor‑β/Smad3 pathway: Is it associated with endoplasmic reticulum stress‑mediated renal interstitial fibrosis?
Molecular medicine reports
Liu Y,Shen P,Zhou Y,Tang L,Chai H
Mon Jul 01 00:00:00 UTC 2019
circ-CSPP1 promotes proliferation, invasion and migration of ovarian cancer cells by acting as a miR-1236-3p sponge.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li QH,Liu Y,Chen S,Zong ZH,Du YP,Sheng XJ,Zhao Y
Sat Jun 01 00:00:00 UTC 2019
Establishment of an Experimental Intracerebral Haemorrhage Model for Mass Effect Research using a Thermo-sensitive Hydrogel.
Scientific reports
Gong Y,Gong Y,Hou Z,Guo T,Deng J,Hao S,Wang B
Wed Sep 25 00:00:00 UTC 2019
Anti-apoptotic effect of the Shh signaling pathway in cigarette smoke extract induced MLE 12 apoptosis.
Tobacco induced diseases
Li J,Zong D,Chen Y,Chen P
Thu Oct 01 00:00:00 UTC 2020
Protective Effects of 18β-Glycyrrhetinic Acid on Monocrotaline-Induced Pulmonary Arterial Hypertension in Rats.
Frontiers in pharmacology
Zhang M,Chang Z,Zhao F,Zhang P,Hao YJ,Yan L,Liu N,Wang JL,Bo L,Ma P,Zhou W,Ma X,Xu QB,Zhou R
Thu Oct 01 00:00:00 UTC 2020
PNS-R1 inhibits Dex-induced bronchial epithelial cells apoptosis in asthma through mitochondrial apoptotic pathway.
Cell & bioscience
Zou W,Niu C,Fu Z,Gong C
Thu Oct 01 00:00:00 UTC 2020
An isopentenyl-substituted flavonoid norartocarpin activates Nrf2 signalling pathway and prevents oxidative insults in human lung epithelial cells.
Free radical research
Yang WJ,He JX,Zhou MX,Huang M,Wang SQ,Wang XN,Lou HX,Ren DM,Shen T
Fri Mar 01 00:00:00 UTC 2019
Mitophagy is a protective response against oxidative damage in bone marrow mesenchymal stem cells.
Life sciences
Fan P,Yu XY,Xie XH,Chen CH,Zhang P,Yang C,Peng X,Wang YT
Mon Jul 15 00:00:00 UTC 2019
Neuroprotective effect of Vanillin on hypoxic-ischemic brain damage in neonatal rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Lan XB,Wang Q,Yang JM,Ma L,Zhang WJ,Zheng P,Sun T,Niu JG,Liu N,Yu JQ
Tue Oct 01 00:00:00 UTC 2019
ROS/KRAS/AMPK Signaling Contributes to Gemcitabine-Induced Stem-like Cell Properties in Pancreatic Cancer.
Molecular therapy oncolytics
Zhao H,Wu S,Li H,Duan Q,Zhang Z,Shen Q,Wang C,Yin T
Fri Sep 27 00:00:00 UTC 2019
GPX2 suppression of H2O2 stress regulates cervical cancer metastasis and apoptosis via activation of the β-catenin-WNT pathway.
OncoTargets and therapy
Wang Y,Cao P,Alshwmi M,Jiang N,Xiao Z,Jiang F,Gu J,Wang X,Sun X,Li S
Mon Apr 11 00:00:00 UTC 2022
The selective HDAC6 inhibitor Nexturastat A induces apoptosis, overcomes drug resistance and inhibits tumor growth in multiple myeloma.
Bioscience reports
Sun X,Xie Y,Sun X,Yao Y,Li H,Li Z,Yao R,Xu K
Fri Mar 29 00:00:00 UTC 2019
Differential immune responses of C57BL/6 mice to infection by Salmonella enterica serovar Typhimurium strain SL1344, CVCC541 and CMCC50115.
Virulence
Wei S,Huang J,Liu Z,Wang M,Zhang B,Lian Z,Guo Y,Han H
Sun Dec 01 00:00:00 UTC 2019
Design, synthesis and molecular mechanisms of novel dual inhibitors of heat shock protein 90/phosphoinositide 3-kinase alpha (Hsp90/PI3Kα) against cutaneous melanoma.
Journal of enzyme inhibition and medicinal chemistry
Qin F,Wang Y,Jiang X,Wang Y,Zhang N,Wen X,Wang L,Jiang Q,He G
Sun Dec 01 00:00:00 UTC 2019
Folic acid delays age-related cognitive decline in senescence-accelerated mouse prone 8: alleviating telomere attrition as a potential mechanism.
Aging
Lv X,Wang X,Wang Y,Zhou D,Li W,Wilson JX,Chang H,Huang G
Fri Nov 22 00:00:00 UTC 2019
The endogenous retrovirus-derived long noncoding RNA TROJAN promotes triple-negative breast cancer progression via ZMYND8 degradation.
Science advances
Jin X,Xu XE,Jiang YZ,Liu YR,Sun W,Guo YJ,Ren YX,Zuo WJ,Hu X,Huang SL,Shen HJ,Lan F,He YF,Hu GH,Di GH,He XH,Li DQ,Liu S,Yu KD,Shao ZM
Fri Mar 01 00:00:00 UTC 2019
Hypoxic BMSC-derived exosomal miRNAs promote metastasis of lung cancer cells via STAT3-induced EMT.
Molecular cancer
Zhang X,Sai B,Wang F,Wang L,Wang Y,Zheng L,Li G,Tang J,Xiang J
Wed Mar 13 00:00:00 UTC 2019
Identification of proteomic signatures associated with COPD frequent exacerbators.
Life sciences
Sun P,Ye R,Wang C,Bai S,Zhao L
Thu Aug 01 00:00:00 UTC 2019
ATXN3 promotes breast cancer metastasis by deubiquitinating KLF4.
Cancer letters
Zou H,Chen H,Zhou Z,Wan Y,Liu Z
Sat Dec 28 00:00:00 UTC 2019
RAD51 is a potential marker for prognosis and regulates cell proliferation in pancreatic cancer.
Cancer cell international
Zhang X,Ma N,Yao W,Li S,Ren Z
Fri Oct 27 00:00:00 UTC 2023
Melatonin protects against COPD by attenuating apoptosis and endoplasmic reticulum stress via upregulating SIRT1 expression in rats.
Canadian journal of physiology and pharmacology
He B,Zhang W,Qiao J,Peng Z,Chai X
Wed May 01 00:00:00 UTC 2019
Carnosol as a Nrf2 Activator Improves Endothelial Barrier Function Through Antioxidative Mechanisms.
International journal of molecular sciences
Li X,Zhang Q,Hou N,Li J,Liu M,Peng S,Zhang Y,Luo Y,Zhao B,Wang S,Zhang Y,Qiao Y
Mon Feb 18 00:00:00 UTC 2019
Induced Pluripotent Stem Cells Regulate Triggering Receptor Expressed on Myeloid Cell-1 Expression and the p38 Mitogen-Activated Protein Kinase Pathway in Endotoxin-Induced Acute Lung Injury.
Stem cells (Dayton, Ohio)
Su VY,Yang KY,Chiou SH,Chen NJ,Mo MH,Lin CS,Wang CT
Wed May 01 00:00:00 UTC 2019
Multi-walled carbon nanotubes (MWCNTs) promoted lipid accumulation in THP-1 macrophages through modulation of endoplasmic reticulum (ER) stress.
Nanotoxicology
Long J,Ma W,Yu Z,Liu H,Cao Y
Sun Sep 01 00:00:00 UTC 2019
SIRT5-mediated SDHA desuccinylation promotes clear cell renal cell carcinoma tumorigenesis.
Free radical biology & medicine
Ma Y,Qi Y,Wang L,Zheng Z,Zhang Y,Zheng J
Mon Apr 01 00:00:00 UTC 2019
MicroRNA‑223 promotes osteoblast differentiation of MC3T3‑E1 cells by targeting histone deacetylase 2.
International journal of molecular medicine
Chen J,He G,Wang Y,Cai D
Fri Mar 01 00:00:00 UTC 2019
Upregulated histamine receptor H3 promotes tumor growth and metastasis in hepatocellular carcinoma.
Oncology reports
Yu D,Zhao J,Wang Y,Hu J,Zhao Q,Li J,Zhu J
Sat Jun 01 00:00:00 UTC 2019
Kallikrein 12 Regulates Innate Resistance of Murine Macrophages against Mycobacterium bovis Infection by Modulating Autophagy and Apoptosis.
Cells
Sabir N,Hussain T,Liao Y,Wang J,Song Y,Shahid M,Cheng G,Mangi MH,Yao J,Yang L,Zhao D,Zhou X
Sun May 05 00:00:00 UTC 2019
CX-F9, a novel RSK2 inhibitor, suppresses cutaneous melanoma cells proliferation and metastasis through regulating autophagy.
Biochemical pharmacology
Zhang X,Cai L,Zhao S,Long J,Li J,Wu L,Su J,Zhang J,Tao J,Zhou J,Chen X,Peng C
Tue Oct 01 00:00:00 UTC 2019
TAK242 suppresses the TLR4 signaling pathway and ameliorates DCD liver IRI in rats.
Molecular medicine reports
Zhong X,Xiao Q,Liu Z,Wang W,Lai CH,Yang W,Yue P,Ye Q,Xiao J
Sun Sep 01 00:00:00 UTC 2019
Propofol Attenuates Hypoxia/Reoxygenation-Induced Apoptosis and Autophagy in HK-2 Cells by Inhibiting JNK Activation.
Yonsei medical journal
Wang H,Peng X,Huang Y,Xiao Y,Wang Z,Zhan L
Sun Dec 01 00:00:00 UTC 2019
Rho-kinase inhibitor attenuates airway mucus hypersecretion and inflammation partly by downregulation of IL-13 and the JNK1/2-AP1 signaling pathway.
Biochemical and biophysical research communications
Zhang W,Li X,Zhang Y
Tue Aug 20 00:00:00 UTC 2019
The Therapeutic Effects of Treadmill Exercise on Osteoarthritis in Rats by Inhibiting the HDAC3/NF-KappaB Pathway in vivo and in vitro.
Frontiers in physiology
Zhang H,Ji L,Yang Y,Wei Y,Zhang X,Gang Y,Lu J,Bai L
Fri Oct 13 00:00:00 UTC 2023
Multi-walled carbon nanotubes promoted lipid accumulation in human aortic smooth muscle cells.
Toxicology and applied pharmacology
Yang H,Li J,Yang C,Liu H,Cao Y
Mon Jul 01 00:00:00 UTC 2019
Clock mutant promotes osteoarthritis by inhibiting the acetylation of NFκB.
Osteoarthritis and cartilage
Yuan G,Xu L,Cai T,Hua B,Sun N,Yan Z,Lu C,Qian R
Sat Jun 01 00:00:00 UTC 2019
Bilirubin alleviates alum-induced peritonitis through inactivation of NLRP3 inflammasome.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Lin Y,Wang S,Yang Z,Gao L,Zhou Z,Yu P,Wu B
Thu Aug 01 00:00:00 UTC 2019
Identification of predictors of drug sensitivity using patient-derived models of esophageal squamous cell carcinoma.
Nature communications
Su D,Zhang D,Jin J,Ying L,Han M,Chen K,Li B,Wu J,Xie Z,Zhang F,Lin Y,Cheng G,Li JY,Huang M,Wang J,Wang K,Zhang J,Li F,Xiong L,Futreal A,Mao W
Thu Nov 07 00:00:00 UTC 2019
RING finger protein 10 is a potential drug target for diabetic vascular complications.
Molecular medicine reports
Li S,Yu G,Huang W,Wang R,Pu P,Chen M
Thu Aug 01 00:00:00 UTC 2019
MiR-664-2 impacts pubertal development in a precocious-puberty rat model through targeting the NMDA receptor-1†.
Biology of reproduction
Ju M,Yang L,Zhu J,Chen Z,Zhang M,Yu J,Tian Z
Sat Jun 01 00:00:00 UTC 2019
MiR-24-3p Inhibits the Progression of Pancreatic Ductal Adenocarcinoma Through LAMB3 Downregulation.
Frontiers in oncology
Huang W,Gu J,Tao T,Zhang J,Wang H,Fan Y
Mon Sep 28 00:00:00 UTC 2020
Structural Characterization and Antioxidant Activity of Polysaccharides from Athyrium multidentatum (Doll.) Ching in d-Galactose-Induced Aging Mice via PI3K/AKT Pathway.
Molecules (Basel, Switzerland)
Jing L,Jiang JR,Liu DM,Sheng JW,Zhang WF,Li ZJ,Wei LY
Mon Sep 16 00:00:00 UTC 2019
NAMPT overexpression alleviates alcohol-induced hepatic steatosis in mice.
PloS one
Xiong X,Yu J,Fan R,Zhang C,Xu L,Sun X,Huang Y,Wang Q,Ruan HB,Qian X
Mon Nov 18 00:00:00 UTC 2019
P11 Loss-of-Function is Associated with Decreased Cell Proliferation and Neurobehavioral Disorders in Mice.
International journal of biological sciences
Liu G,Wang Y,Zheng W,Cheng H,Zhou R
Tue Apr 07 00:00:00 UTC 2020
Deciphering novel biomarkers of lymph node metastasis of thyroid papillary microcarcinoma using proteomic analysis of ultrasound-guided fine-needle aspiration biopsy samples.
Journal of proteomics
Lin P,Yao Z,Sun Y,Li W,Liu Y,Liang K,Liu Y,Qin J,Hou X,Chen L
Tue Jul 30 00:00:00 UTC 2019
Chrysanthemum extract attenuates hepatotoxicity via inhibiting oxidative stress in vivo and in vitro.
Food & nutrition research
Tian Z,Jia H,Jin Y,Wang M,Kou J,Wang C,Rong X,Xie X,Han G,Pang X
Fri Apr 08 00:00:00 UTC 2022
High metabolic rate and stem cell characteristics of esophageal cancer stem-like cells depend on the Hsp27-AKT-HK2 pathway.
International journal of cancer
Liu CC,Chou KT,Hsu JW,Lin JH,Hsu TW,Yen DH,Hung SC,Hsu HS
Tue Oct 15 00:00:00 UTC 2019
Turnover of bile acids in liver, serum and caecal content by high-fat diet feeding affects hepatic steatosis in rats.
Biochimica et biophysica acta. Molecular and cell biology of lipids
Tang Y,Zhang J,Li J,Lei X,Xu D,Wang Y,Li C,Li X,Mao Y
Tue Oct 01 00:00:00 UTC 2019
Knockdown of LncRNA SCAMP1 suppressed malignant biological behaviours of glioma cells via modulating miR-499a-5p/LMX1A/NLRC5 pathway.
Journal of cellular and molecular medicine
Zong Z,Song Y,Xue Y,Ruan X,Liu X,Yang C,Zheng J,Cao S,Li Z,Liu Y
Thu Aug 01 00:00:00 UTC 2019
CCL18 promotes the metastasis of squamous cell carcinoma of the head and neck through MTDH-NF-κB signalling pathway.
Journal of cellular and molecular medicine
Qin Y,Wang J,Zhu G,Li G,Tan H,Chen C,Pi L,She L,Chen X,Wei M,Li Z,Liu Z,Huang D,Liu Y,Zhang X
Mon Apr 01 00:00:00 UTC 2019
Cell survival controlled by lens-derived Sema3A-Nrp1 is vital on caffeine-suppressed corneal innervation during chick organogenesis.
Journal of cellular physiology
Wang G,Jiang LM,Tan BY,Li PZ,Zhang PL,Zhang Y,Cheng X,Ma ZL,Li ZJ,Brand-Saberi B,Yang X
Sat Jun 01 00:00:00 UTC 2019
Inhibition of soluble epoxide hydrolase ameliorates hyperhomocysteinemia-induced hepatic steatosis by enhancing β-oxidation of fatty acid in mice.
American journal of physiology. Gastrointestinal and liver physiology
Yao L,Cao B,Cheng Q,Cai W,Ye C,Liang J,Liu W,Tan L,Yan M,Li B,He J,Hwang SH,Zhang X,Wang C,Ai D,Hammock BD,Zhu Y
Mon Apr 01 00:00:00 UTC 2019
Changes among TGF-β1+ Breg cells and helper T cell subsets in a murine model of allergic rhinitis with prolonged OVA challenge.
International immunopharmacology
Wang M,Gu Z,Yang J,Zhao H,Cao Z
Mon Apr 01 00:00:00 UTC 2019
Induction of Endoplasmic Reticulum Stress by Cadmium and Its Regulation on Nrf2 Signaling Pathway in Kidneys of Rats.
Biomedical and environmental sciences : BES
Chen ZJ,Chen JX,Wu LK,Li BY,Tian YF,Xian M,Huang ZP,Yu RA
Tue Jan 01 00:00:00 UTC 2019
DHT deteriorates the progression of monocrotaline-induced pulmonary arterial hypertension: effects of endogenous and exogenous androgen.
American journal of translational research
Wen J,Wang J,Tang X,Deng S,Dai J,Li X,Cai W
Thu Oct 01 00:00:00 UTC 2020
Protective mechanism of Taxifolin for chlorpyrifos neurotoxicity in BV2 cells.
Neurotoxicology
Zhang C,Zhan J,Zhao M,Dai H,Deng Y,Zhou W,Zhao L
Sun Sep 01 00:00:00 UTC 2019
Phenotypic transition of tumor cells between epithelial- and mesenchymal-like state during adaptation to acidosis.
Cell cycle (Georgetown, Tex.)
Zhang Y,Xu L,Wang P,Jian H,Shi X,Jia M,Mo L,Hu Z,Li H,Li J
Thu Aug 01 00:00:00 UTC 2019
Paip1 Indicated Poor Prognosis in Cervical Cancer and Promoted Cervical Carcinogenesis.
Cancer research and treatment
Li N,Piao J,Wang X,Kim KY,Bae JY,Ren X,Lin Z
Tue Oct 01 00:00:00 UTC 2019
Nerve injury elevates functional Cav3.2 channels in superficial spinal dorsal horn.
Molecular pain
Feng XJ,Ma LX,Jiao C,Kuang HX,Zeng F,Zhou XY,Cheng XE,Zhu MY,Zhang DY,Jiang CY,Liu T
Mon Aug 12 00:00:00 UTC 2019
RNA-Seq analysis of potential lncRNAs and genes for the anti-renal fibrotic effect of norcantharidin.
Journal of cellular biochemistry
Xiao H,Liao Y,Tang C,Xiao Z,Luo H,Li J,Liu H,Sun L,Zeng D,Li Y
Tue Oct 01 00:00:00 UTC 2019
Iron dysregulation in vascular dementia: Focused on the AMPK/autophagy pathway.
Brain research bulletin
Huo T,Jia Y,Yin C,Luo X,Zhao J,Wang Z,Lv P
Fri Nov 01 00:00:00 UTC 2019
Chitosan Ameliorates DSS-Induced Ulcerative Colitis Mice by Enhancing Intestinal Barrier Function and Improving Microflora.
International journal of molecular sciences
Wang J,Zhang C,Guo C,Li X
Fri Nov 15 00:00:00 UTC 2019
Dose-Dependent Effects of Triclocarban Exposure on Lipid Homeostasis in Rats.
Chemical research in toxicology
Dong M,Xu X,Huang Q,Lei H,Xu G,Ma J,Hatzakis E,Wang X,Zhang L
Mon Nov 18 00:00:00 UTC 2019
Glycolytic Enzyme PKM2 Mediates Autophagic Activation to Promote Cell Survival in NPM1-Mutated Leukemia.
International journal of biological sciences
Wang L,Yang L,Yang Z,Tang Y,Tao Y,Zhan Q,Lei L,Jing Y,Jiang X,Jin H,Zou Q,Xian J,Zhang L
Mon Feb 03 00:00:00 UTC 2020
Protrudin modulates seizure activity through GABAA receptor regulation.
Cell death & disease
Lu X,Yang Y,Zhou R,Li Y,Yang Y,Wang X
Wed Nov 27 00:00:00 UTC 2019
Dietary Curcumin Supplementation Increases Antioxidant Capacity, Upregulates Nrf2 and Hmox1 Levels in the Liver of Piglet Model with Intrauterine Growth Retardation.
Nutrients
Niu Y,He J,Ahmad H,Shen M,Zhao Y,Gan Z,Zhang L,Zhong X,Wang C,Wang T
Thu Dec 05 00:00:00 UTC 2019
Porous Se@SiO2 nanocomposite promotes migration and osteogenic differentiation of rat bone marrow mesenchymal stem cell to accelerate bone fracture healing in a rat model.
International journal of nanomedicine
Li C,Wang Q,Gu X,Kang Y,Zhang Y,Hu Y,Li T,Jin H,Deng G,Wang Q
Thu Aug 08 00:00:00 UTC 2019
Mild hypothermia improves neurological outcome in mice after cardiopulmonary resuscitation through Silent Information Regulator 1-actviated autophagy.
Cell death discovery
Wei H,Yin M,Lu Y,Yang Y,Li B,Liao XX,Dai G,Jing X,Xiong Y,Hu C
Thu Oct 01 00:00:00 UTC 2020
Abnormal serum vitamin A levels and retinoic acid receptor α expression patterns in children with anorectal malformation.
Pediatric surgery international
Wang Z,Wang Q,Gu C,Zhang J,Wang Y
Thu Aug 01 00:00:00 UTC 2019
Naringenin Produces Neuroprotection Against LPS-Induced Dopamine Neurotoxicity via the Inhibition of Microglial NLRP3 Inflammasome Activation.
Frontiers in immunology
Chen C,Wei YZ,He XM,Li DD,Wang GQ,Li JJ,Zhang F
Wed Sep 09 00:00:00 UTC 2020
Association between CIRP expression and hypoxic-ischemic brain injury in neonatal rats.
Experimental and therapeutic medicine
Chen L,Tian Q,Wang W
Sun Sep 01 00:00:00 UTC 2019
Osteoadherin serves roles in the regulation of apoptosis and growth in MC3T3‑E1 osteoblast cells.
International journal of molecular medicine
Hamaya E,Fujisawa T,Tamura M
Sun Dec 01 00:00:00 UTC 2019
Quantitative proteomic profiling of tumor-associated vascular endothelial cells in colorectal cancer.
Biology open
Wang G,Yang Q,Li M,Zhang Y,Cai Y,Liang X,Fu Y,Xiao Z,Zhou M,Xie Z,Huang H,Huang Y,Chen Y,He Q,Peng F,Chen Z
Mon May 13 00:00:00 UTC 2019
Brilliant Blue G Inhibits Inflammasome Activation and Reduces Disruption of Blood-Spinal Cord Barrier Induced by Spinal Cord Injury in Rats.
Medical science monitor : international medical journal of experimental and clinical research
Zhou X,Yang Y,Wu L,Wang Y,Du C,Li C,Wang Z,Wang Y
Sat Aug 24 00:00:00 UTC 2019
Targeting Exosomal EBV-LMP1 Transfer and miR-203 Expression via the NF-κB Pathway: The Therapeutic Role of Aspirin in NPC.
Molecular therapy. Nucleic acids
Zuo L,Xie Y,Tang J,Xin S,Liu L,Zhang S,Yan Q,Zhu F,Lu J
Fri Sep 06 00:00:00 UTC 2019
Autophagy Attenuation Hampers Progesterone Synthesis during the Development of Pregnant Corpus Luteum.
Cells
Tang Z,Zhang Z,Zhang H,Wang Y,Zhang Y,Zhao J,Yang H,Wang Z
Fri Dec 27 00:00:00 UTC 2019
Nilotinib: A Tyrosine Kinase Inhibitor Mediates Resistance to Intracellular Mycobacterium Via Regulating Autophagy.
Cells
Hussain T,Zhao D,Shah SZA,Sabir N,Wang J,Liao Y,Song Y,Dong H,Hussain Mangi M,Ni J,Yang L,Zhou X
Sun May 26 00:00:00 UTC 2019
The methionine salvage pathway-involving ADI1 inhibits hepatoma growth by epigenetically altering genes expression via elevating S-adenosylmethionine.
Cell death & disease
Chu YD,Lai HY,Pai LM,Huang YH,Lin YH,Liang KH,Yeh CT
Mon Mar 11 00:00:00 UTC 2019
Hepatitis B Virus X Protein-Induced RORγ Expression to Promote the Migration and Proliferation of Hepatocellular Carcinoma.
BioMed research international
Huang Y,Liang H,He C,Peng F
Mon Apr 27 00:00:00 UTC 2020
Eutopic endometrium from patients with endometriosis modulates the expression of CD36 and SIRP-α in peritoneal macrophages.
The journal of obstetrics and gynaecology research
Xie Q,He H,Wu YH,Zou LJ,She XL,Xia XM,Wu XQ
Wed May 01 00:00:00 UTC 2019
Neuroprotective Effect of Swertiamain on Cerebral Ischemia/Reperfusion Injury by Inducing the Nrf2 Protective Pathway.
ACS chemical neuroscience
Wang H,Wei W,Lan X,Liu N,Li Y,Ma H,Sun T,Peng X,Zhuang C,Yu J
Wed May 15 00:00:00 UTC 2019
Identification of diterpenoid compounds that interfere with Fli-1 DNA binding to suppress leukemogenesis.
Cell death & disease
Liu T,Xia L,Yao Y,Yan C,Fan Y,Gajendran B,Yang J,Li YJ,Chen J,Filmus J,Spaner DE,Zacksenhaus E,Hao X,Ben-David Y
Mon Feb 11 00:00:00 UTC 2019
The lncRNA HOXA11-AS regulates Rab3D expression by sponging miR-125a-5p promoting metastasis of osteosarcoma.
Cancer management and research
Cao K,Fang Y,Wang H,Jiang Z,Guo L,Hu Y
Fri Jan 07 00:00:00 UTC 2022
Site-Specific Immobilization of β2-AR Using O6-Benzylguanine Derivative-Functionalized Supporter for High-Throughput Receptor-Targeting Lead Discovery.
Analytical chemistry
Wang J,Wang Y,Liu J,Li Q,Yin G,Zhang Y,Xiao C,Fan T,Zhao X,Zheng X
Tue Jun 04 00:00:00 UTC 2019
LncRNA EPB41L4A-AS1 regulates glycolysis and glutaminolysis by mediating nucleolar translocation of HDAC2.
EBioMedicine
Liao M,Liao W,Xu N,Li B,Liu F,Zhang S,Wang Y,Wang S,Zhu Y,Chen D,Xie W,Jiang Y,Cao L,Yang BB,Zhang Y
Fri Mar 01 00:00:00 UTC 2019
The role of autophagy in the overexpression of MUC5AC in patients with chronic rhinosinusitis.
International immunopharmacology
Ye Y,Zhao J,Ye J,Jiang X,Liu H,Xie Y,Zhang J,Luo Q
Sat Jun 01 00:00:00 UTC 2019
Transketolase (TKT) activity and nuclear localization promote hepatocellular carcinoma in a metabolic and a non-metabolic manner.
Journal of experimental & clinical cancer research : CR
Qin Z,Xiang C,Zhong F,Liu Y,Dong Q,Li K,Shi W,Ding C,Qin L,He F
Thu Apr 11 00:00:00 UTC 2019
Circular RNA cMras inhibits lung adenocarcinoma progression via modulating miR-567/PTPRG regulatory pathway.
Cell proliferation
Yu C,Tian F,Liu J,Su M,Wu M,Zhu X,Qian W
Wed May 01 00:00:00 UTC 2019
Long Noncoding RNA FAM225A Promotes Nasopharyngeal Carcinoma Tumorigenesis and Metastasis by Acting as ceRNA to Sponge miR-590-3p/miR-1275 and Upregulate ITGB3.
Cancer research
Zheng ZQ,Li ZX,Zhou GQ,Lin L,Zhang LL,Lv JW,Huang XD,Liu RQ,Chen F,He XJ,Kou J,Zhang J,Wen X,Li YQ,Ma J,Liu N,Sun Y
Sun Sep 15 00:00:00 UTC 2019
Propionate relieves pentylenetetrazol-induced seizures, consequent mitochondrial disruption, neuron necrosis and neurological deficits in mice.
Biochemical pharmacology
Cheng Y,Mai Q,Zeng X,Wang H,Xiao Y,Tang L,Li J,Zhang Y,Ding H
Fri Nov 01 00:00:00 UTC 2019
Silencing ubiquitin-conjugating enzyme 2C inhibits proliferation and epithelial-mesenchymal transition in pancreatic ductal adenocarcinoma.
The FEBS journal
Wang X,Yin L,Yang L,Zheng Y,Liu S,Yang J,Cui H,Wang H
Sun Dec 01 00:00:00 UTC 2019
NS38 is required for aquareovirus replication via interaction with viral core proteins and host eIF3A.
Virology
Zhang J,Guo H,Zhang F,Chen Q,Chang M,Fang Q
Fri Mar 01 00:00:00 UTC 2019
Time-dependent C5a and C5aR expression in dental pulp cells following stimulation with LTA and LPS.
International journal of molecular medicine
Liu M,Mu H,Peng W,Zhao L,Hu W,Jiang Z,Gao L,Cao X,Li N,Han J
Sun Sep 01 00:00:00 UTC 2019
Netrin‑1 interference potentiates epithelial‑to‑mesenchymal transition through the PI3K/AKT pathway under the hypoxic microenvironment conditions of non‑small cell lung cancer.
International journal of oncology
Jin X,Luan H,Chai H,Yan L,Zhang J,Wang Q,Cao L
Mon Apr 01 00:00:00 UTC 2019
Inhibition of Bcl-2 Synergistically Enhances the Antileukemic Activity of Midostaurin and Gilteritinib in Preclinical Models of FLT3-Mutated Acute Myeloid Leukemia.
Clinical cancer research : an official journal of the American Association for Cancer Research
Ma J,Zhao S,Qiao X,Knight T,Edwards H,Polin L,Kushner J,Dzinic SH,White K,Wang G,Zhao L,Lin H,Wang Y,Taub JW,Ge Y
Fri Nov 15 00:00:00 UTC 2019
p66Shc Contributes to Liver Fibrosis through the Regulation of Mitochondrial Reactive Oxygen Species.
Theranostics
Zhao Y,Wang Z,Feng D,Zhao H,Lin M,Hu Y,Zhang N,Lv L,Gao Z,Zhai X,Tian X,Yao J
Fri Jan 31 00:00:00 UTC 2020
MiR-223-3p functions as a tumor suppressor in lung squamous cell carcinoma by miR-223-3p-mutant p53 regulatory feedback loop.
Journal of experimental & clinical cancer research : CR
Luo P,Wang Q,Ye Y,Zhang J,Lu D,Cheng L,Zhou H,Xie M,Wang B
Tue Feb 12 00:00:00 UTC 2019
Novel ADAM-17 inhibitor ZLDI-8 inhibits the proliferation and metastasis of chemo-resistant non-small-cell lung cancer by reversing Notch and epithelial mesenchymal transition in vitro and in vivo.
Pharmacological research
Lu HY,Zu YX,Jiang XW,Sun XT,Liu TY,Li RL,Wu Q,Zhang YS,Zhao QC
Tue Oct 01 00:00:00 UTC 2019
Transection of the cervical sympathetic trunk inhibits the progression of pulmonary arterial hypertension via ERK-1/2 Signalling.
Respiratory research
Zhao Y,Xiang R,Peng X,Dong Q,Li D,Yu G,Xiao L,Qin S,Huang W
Fri Jun 14 00:00:00 UTC 2019
The toxicity of multi-walled carbon nanotubes (MWCNTs) to human endothelial cells: The influence of diameters of MWCNTs.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Zhao X,Chang S,Long J,Li J,Li X,Cao Y
Mon Apr 01 00:00:00 UTC 2019
Adipose tissue-derived omentin-1 attenuates arterial calcification via AMPK/Akt signaling pathway.
Aging
Xu F,Li FX,Lin X,Zhong JY,Wu F,Shan SK,Tan CM,Yuan LQ,Liao XB
Fri Oct 25 00:00:00 UTC 2019
Metabolism-induced tumor activator 1 (MITA1), an Energy Stress-Inducible Long Noncoding RNA, Promotes Hepatocellular Carcinoma Metastasis.
Hepatology (Baltimore, Md.)
Ma M,Xu H,Liu G,Wu J,Li C,Wang X,Zhang S,Xu H,Ju S,Cheng W,Dai L,Wei Y,Tian Y,Fu X
Mon Jul 01 00:00:00 UTC 2019
Pseudogene RACGAP1P activates RACGAP1/Rho/ERK signalling axis as a competing endogenous RNA to promote hepatocellular carcinoma early recurrence.
Cell death & disease
Wang MY,Chen DP,Qi B,Li MY,Zhu YY,Yin WJ,He L,Yu Y,Li ZY,Lin L,Yang F,Lin ZR,Liu JQ
Mon Jun 03 00:00:00 UTC 2019
Functional analysis of the CXCR1a gene response to SGIV viral infection in grouper.
Fish & shellfish immunology
Wang Q,Yang M,Li C,Wang S,Wang Y,Lin F,Zheng L,Yu Y,Qin Q
Wed May 01 00:00:00 UTC 2019
SIRT6/HIF-1α axis promotes papillary thyroid cancer progression by inducing epithelial-mesenchymal transition.
Cancer cell international
Yang Z,Yu W,Huang R,Ye M,Min Z
Wed Aug 23 00:00:00 UTC 2023
GSTO1 acts as a mediator in sodium fluoride-induced alterations of learning and memory related factors expressions in the hippocampus cell line.
Chemosphere
Wang J,Gao Y,Cheng X,Yang J,Zhao Y,Xu H,Zhu Y,Yan Z,Manthari RK,Ommati MM,Wang J
Mon Jul 01 00:00:00 UTC 2019
Macrophage migration inhibitory factor promotes the invasion and metastasis of oral squamous cell carcinoma through matrix metalloprotein-2/9.
Molecular carcinogenesis
Wang SS,Zheng M,Pang X,Zhang M,Yu XH,Wu JB,Gao XL,Wu JS,Yang X,Tang YJ,Tang YL,Liang XH
Tue Oct 01 00:00:00 UTC 2019
Ghrelin regulates sepsis‑induced rat acute gastric injury.
Molecular medicine reports
Li B,Lin Q,Guo H,Liu L,Li Y
Sat Jun 01 00:00:00 UTC 2019
Downregulation of nicotinamide N-methyltransferase inhibits migration and epithelial-mesenchymal transition of esophageal squamous cell carcinoma via Wnt/β-catenin pathway.
Molecular and cellular biochemistry
Cui Y,Zhang L,Wang W,Ma S,Liu H,Zang X,Zhang Y,Guan F
Tue Oct 01 00:00:00 UTC 2019
Effects of agomelatine in rotenone-induced Parkinson's disease in rats.
Neuroscience letters
Günaydın C,Avcı B,Bozkurt A,Önger ME,Balcı H,Bilge SS
Tue Apr 23 00:00:00 UTC 2019
The interference of DEHP in precocious puberty of females mediated by the hypothalamic IGF-1/PI3K/Akt/mTOR signaling pathway.
Ecotoxicology and environmental safety
Shao P,Wang Y,Zhang M,Wen X,Zhang J,Xu Z,Hu M,Jiang J,Liu T
Tue Oct 15 00:00:00 UTC 2019
Whole-Exome Sequencing Analysis Identified Novel Mutations in the TSPAN12 Gene in Chinese Families with Familial Exudative Vitreoretinopathy.
Genetic testing and molecular biomarkers
Yuan Y,Xu H,Zhang S,Zhang X,Zhang L,Yang Z
Tue Oct 01 00:00:00 UTC 2019
Hepatoprotective effects of zinc (II) via cytochrome P-450/reactive oxygen species and canonical apoptosis pathways after arsenite waterborne exposure in common carp.
Chemosphere
Zhao H,Wang Y,Guo M,Fei D,Mu M,Yu H,Xing M
Sun Dec 01 00:00:00 UTC 2019
Dietary Dihydroartemisinin Supplementation Attenuates Hepatic Oxidative Damage of Weaned Piglets with Intrauterine Growth Retardation through the Nrf2/ARE Signaling Pathway.
Animals : an open access journal from MDPI
Zhao Y,Niu Y,He J,Zhang L,Wang C,Wang T
Fri Dec 13 00:00:00 UTC 2019
Epidural adipose tissue-derived mesenchymal stem cell activation induced by lung cancer cells promotes malignancy and EMT of lung cancer.
Stem cell research & therapy
Wang Y,Chu Y,Ren X,Xiang H,Xi Y,Ma X,Zhu K,Guo Z,Zhou C,Zhang G,Chen B
Thu Jun 13 00:00:00 UTC 2019
Oroxin B Induces Apoptosis by Down-Regulating MicroRNA-221 Resulting in the Inactivation of the PTEN/PI3K/AKT Pathway in Liver Cancer.
Molecules (Basel, Switzerland)
Li N,Men W,Zheng Y,Wang H,Meng X
Sat Nov 30 00:00:00 UTC 2019
Primary Cilia Blockage Promotes the Malignant Behaviors of Hepatocellular Carcinoma via Induction of Autophagy.
BioMed research international
Liu L,Sheng JQ,Wang MR,Gan Y,Wu XL,Liao JZ,Tian DA,He XX,Li PY
Thu Mar 19 00:00:00 UTC 2020
High expression of SMARCE1 predicts poor prognosis and promotes cell growth and metastasis in gastric cancer.
Cancer management and research
Liu H,Zhao YR,Chen B,Ge Z,Huang JS
Thu Oct 01 00:00:00 UTC 2020
Combined melatonin and poricoic acid A inhibits renal fibrosis through modulating the interaction of Smad3 and β-catenin pathway in AKI-to-CKD continuum.
Therapeutic advances in chronic disease
Chen DQ,Cao G,Zhao H,Chen L,Yang T,Wang M,Vaziri ND,Guo Y,Zhao YY
Sun Apr 10 00:00:00 UTC 2022
Molecular mechanism of miR-204 regulates proliferation, apoptosis and autophagy of cervical cancer cells by targeting ATF2.
Artificial cells, nanomedicine, and biotechnology
Li N,Guo X,Liu L,Wang L,Cheng R
Sun Dec 01 00:00:00 UTC 2019
Dexmedetomidine inhibits neuronal apoptosis by inducing Sigma-1 receptor signaling in cerebral ischemia-reperfusion injury.
Aging
Zhai M,Liu C,Li Y,Zhang P,Yu Z,Zhu H,Zhang L,Zhang Q,Wang J,Wang J
Mon Nov 04 00:00:00 UTC 2019
Antitumor Effects of Berberine on Gliomas via Inactivation of Caspase-1-Mediated IL-1β and IL-18 Release.
Frontiers in oncology
Tong L,Xie C,Wei Y,Qu Y,Liang H,Zhang Y,Xu T,Qian X,Qiu H,Deng H
Thu Oct 01 00:00:00 UTC 2020
TLR4/NF-κB Signaling Induces GSDMD-Related Pyroptosis in Tubular Cells in Diabetic Kidney Disease.
Frontiers in endocrinology
Wang Y,Zhu X,Yuan S,Wen S,Liu X,Wang C,Qu Z,Li J,Liu H,Sun L,Liu F
Wed Oct 23 00:00:00 UTC 2019
A novel tumor suppressor SPINK5 targets Wnt/β-catenin signaling pathway in esophageal cancer.
Cancer medicine
Wang Q,Lv Q,Bian H,Yang L,Guo KL,Ye SS,Dong XF,Tao LL
Wed May 01 00:00:00 UTC 2019
Apatinib inhibits glycolysis by suppressing the VEGFR2/AKT1/SOX5/GLUT4 signaling pathway in ovarian cancer cells.
Cellular oncology (Dordrecht)
Chen L,Cheng X,Tu W,Qi Z,Li H,Liu F,Yang Y,Zhang Z,Wang Z
Tue Oct 01 00:00:00 UTC 2019
Long non-coding RNA MAGI2-AS3 inhibits breast cancer cell migration and invasion via sponging microRNA-374a.
Cancer biomarkers : section A of Disease markers
Du S,Hu W,Zhao Y,Zhou H,Wen W,Xu M,Zhao P,Liu K
Mon Aug 05 00:00:00 UTC 2019
Niban protein regulates apoptosis in HK-2 cells via caspase-dependent pathway.
Renal failure
Tang S,Wang J,Liu J,Huang Y,Zhou Y,Yang S,Zhang W,Yang M,Zhang H
Fri Nov 01 00:00:00 UTC 2019
Fluorene-9-bisphenol inhibits epithelial-mesenchymal transition of human endometrial cancer Ishikawa cells by repressing TGF-β signaling pathway.
Environmental science and pollution research international
Wang L,Zhuang T,Li F,Wei W
Sun Sep 01 00:00:00 UTC 2019
Critical role of Tim-3 mediated autophagy in chronic stress induced immunosuppression.
Cell & bioscience
Qin A,Zhong T,Zou H,Wan X,Yao B,Zheng X,Yin D
Thu Mar 31 00:00:00 UTC 2022
miR-23a-5p inhibits cell proliferation and invasion in pancreatic ductal adenocarcinoma by suppressing ECM1 expression.
American journal of translational research
Huang W,Huang Y,Gu J,Zhang J,Yang J,Liu S,Xie C,Fan Y,Wang H
Tue Sep 29 00:00:00 UTC 2020
UHRF1 Promotes Proliferation of Human Adipose-Derived Stem Cells and Suppresses Adipogenesis via Inhibiting Peroxisome Proliferator-Activated Receptor γ.
BioMed research international
Chen K,Guo Z,Luo Y,Yuan J,Mo Z
Wed Jan 15 00:00:00 UTC 2020
Bronchial epithelial cell extracellular vesicles ameliorate epithelial-mesenchymal transition in COPD pathogenesis by alleviating M2 macrophage polarization.
Nanomedicine : nanotechnology, biology, and medicine
He S,Chen D,Hu M,Zhang L,Liu C,Traini D,Grau GE,Zeng Z,Lu J,Zhou G,Xie L,Sun S
Sat Jun 01 00:00:00 UTC 2019
PPAR-δ of orange-spotted grouper exerts antiviral activity against fish virus and regulates interferon signaling and inflammatory factors.
Fish & shellfish immunology
Wang Y,Yu Y,Wang Q,Wei S,Wang S,Qin Q,Yang M
Fri Nov 01 00:00:00 UTC 2019
Jiawei Foshou San Induces Apoptosis in Ectopic Endometrium Based on Systems Pharmacology, Molecular Docking, and Experimental Evidence.
Evidence-based complementary and alternative medicine : eCAM
Wei J,Zhao B,Zhang C,Shen B,Zhang Y,Li C,Chen Y
Thu Oct 01 00:00:00 UTC 2020
Casticin inhibits nasopharyngeal carcinoma growth by targeting phosphoinositide 3-kinase.
Cancer cell international
Liu J,Yang J,Hou Y,Zhu Z,He J,Zhao H,Ye X,Li D,Wu Z,Huang Z,Hao B,Yao K
Wed Apr 13 00:00:00 UTC 2022
The Binding of PD-L1 and Akt Facilitates Glioma Cell Invasion Upon Starvation via Akt/Autophagy/F-Actin Signaling.
Frontiers in oncology
Chen RQ,Xu XH,Liu F,Li CY,Li YJ,Li XR,Jiang GY,Hu F,Liu D,Pan F,Qiu XY,Chen XQ
Thu Oct 01 00:00:00 UTC 2020
Icariin targets Nrf2 signaling to inhibit microglia-mediated neuroinflammation.
International immunopharmacology
Zheng Y,Zhu G,He J,Wang G,Li D,Zhang F
Thu Aug 01 00:00:00 UTC 2019
Protective Effect of Koumine, an Alkaloid from Gelsemium Sempervirens, on Injury Induced by H₂O₂ in IPEC-J2 Cells.
International journal of molecular sciences
Yuan Z,Liang Z,Yi J,Chen X,Li R,Wu Y,Wu J,Sun Z
Mon Feb 11 00:00:00 UTC 2019
LncRNA expression profile during autophagy and Malat1 function in macrophages.
PloS one
Ma Z,Zhang J,Xu X,Qu Y,Dong H,Dang J,Huo Z,Xu G
Thu Apr 02 00:00:00 UTC 2020
YWHA/14-3-3 proteins recognize phosphorylated TFEB by a noncanonical mode for controlling TFEB cytoplasmic localization.
Autophagy
Xu Y,Ren J,He X,Chen H,Wei T,Feng W
Sat Jun 01 00:00:00 UTC 2019
HIF-1α contributes to Ang II-induced inflammatory cytokine production in podocytes.
BMC pharmacology & toxicology
Huang H,Fan Y,Gao Z,Wang W,Shao N,Zhang L,Yang Y,Zhu W,Chen Z,Hu J,Ding G
Thu Oct 17 00:00:00 UTC 2019
Jiedu Sangen Decoction Inhibits the Invasion and Metastasis of Colorectal Cancer Cells by Regulating EMT through the Hippo Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Yuan L,Zhou M,Wasan HS,Zhang K,Li Z,Guo K,Shen F,Shen M,Ruan S
Wed Sep 30 00:00:00 UTC 2020
Protective effects of pyrroloquinoline quinine against oxidative stress-induced cellular senescence and inflammation in human renal tubular epithelial cells via Keap1/Nrf2 signaling pathway.
International immunopharmacology
Wang Z,Han N,Zhao K,Li Y,Chi Y,Wang B
Mon Jul 01 00:00:00 UTC 2019
Catalpol Exerts a Neuroprotective Effect in the MPTP Mouse Model of Parkinson's Disease.
Frontiers in aging neuroscience
Wang LY,Yu X,Li XX,Zhao YN,Wang CY,Wang ZY,He ZY
Thu Oct 01 00:00:00 UTC 2020
Exosomes from high glucose-treated macrophages activate glomerular mesangial cells via TGF-β1/Smad3 pathway in vivo and in vitro.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhu QJ,Zhu M,Xu XX,Meng XM,Wu YG
Thu Aug 01 00:00:00 UTC 2019
Microglial NLRP3 inflammasome activation mediates IL-1β release and contributes to central sensitization in a recurrent nitroglycerin-induced migraine model.
Journal of neuroinflammation
He W,Long T,Pan Q,Zhang S,Zhang Y,Zhang D,Qin G,Chen L,Zhou J
Wed Apr 10 00:00:00 UTC 2019
Bone Mesenchymal Stem Cell-Derived Extracellular Vesicles Promote Recovery Following Spinal Cord Injury via Improvement of the Integrity of the Blood-Spinal Cord Barrier.
Frontiers in neuroscience
Lu Y,Zhou Y,Zhang R,Wen L,Wu K,Li Y,Yao Y,Duan R,Jia Y
Thu Oct 01 00:00:00 UTC 2020
Inhibition of Mitochondrial Complex I Aggravates Folic Acid-Induced Acute Kidney Injury.
Kidney & blood pressure research
Zhang W,Yang Y,Gao H,Zhang Y,Jia Z,Huang S
Tue Mar 03 00:00:00 UTC 2020
Activation of Nrf2 signaling by Icariin protects against 6-OHDA-induced neurotoxicity.
Biotechnology and applied biochemistry
Zhu L,Li D,Chen C,Wang G,Shi J,Zhang F
Wed May 01 00:00:00 UTC 2019
Bone marrow mesenchymal stem cells could reduce the toxic effects of hexavalent chromium on the liver by decreasing endoplasmic reticulum stress-mediated apoptosis via SIRT1/HIF-1α signaling pathway in rats.
Toxicology letters
Zhao Y,Yan J,Li AP,Zhang ZL,Li ZR,Guo KJ,Zhao KC,Ruan Q,Guo L
Thu Aug 01 00:00:00 UTC 2019
BPTF promotes hepatocellular carcinoma growth by modulating hTERT signaling and cancer stem cell traits.
Redox biology
Zhao X,Zheng F,Li Y,Hao J,Tang Z,Tian C,Yang Q,Zhu T,Diao C,Zhang C,Chen M,Hu S,Guo P,Zhang L,Liao Y,Yu W,Chen M,Zou L,Guo W,Deng W
Tue Jan 01 00:00:00 UTC 2019
BPTF promotes hepatocellular carcinoma growth by modulating hTERT signaling and cancer stem cell traits.
Redox biology
Zhao X,Zheng F,Li Y,Hao J,Tang Z,Tian C,Yang Q,Zhu T,Diao C,Zhang C,Chen M,Hu S,Guo P,Zhang L,Liao Y,Yu W,Chen M,Zou L,Guo W,Deng W
Tue Jan 01 00:00:00 UTC 2019
Selective ERK1/2 agonists isolated from Melia azedarach with potent anti-leukemic activity.
BMC cancer
Wang N,Fan Y,Yuan CM,Song J,Yao Y,Liu W,Gajendran B,Zacksenhaus E,Li Y,Liu J,Hao XJ,Ben-David Y
Fri Aug 02 00:00:00 UTC 2019
Effect of hydrogen-rich water on the Nrf2/ARE signaling pathway in rats with myocardial ischemia-reperfusion injury.
Journal of bioenergetics and biomembranes
Li L,Liu T,Liu L,Li S,Zhang Z,Zhang R,Zhou Y,Liu F
Sun Dec 01 00:00:00 UTC 2019
MTHFD1L-Mediated Redox Homeostasis Promotes Tumor Progression in Tongue Squamous Cell Carcinoma.
Frontiers in oncology
Li H,Fu X,Yao F,Tian T,Wang C,Yang A
Thu Oct 01 00:00:00 UTC 2020
A novel long noncoding RNA PGC1β-OT1 regulates adipocyte and osteoblast differentiation through antagonizing miR-148a-3p.
Cell death and differentiation
Yuan H,Xu X,Feng X,Zhu E,Zhou J,Wang G,Tian L,Wang B
Tue Oct 01 00:00:00 UTC 2019
Targeting β2 subunit of Na+/K+-ATPase induces glioblastoma cell apoptosis through elevation of intracellular Ca2.
American journal of cancer research
Li S,Dai Z,Yang D,Li W,Dai H,Sun B,Liu X,Xie X,Xu R,Zhao X
Thu Oct 12 00:00:00 UTC 2023
Piperine as a neuroprotective functional component in rats with cerebral ischemic injury.
Food science & nutrition
Hua S,Liu J,Zhang Y,Li J,Zhang X,Dong L,Zhao Y,Fu X
Fri Nov 01 00:00:00 UTC 2019
CD36 plays a negative role in the regulation of lipophagy in hepatocytes through an AMPK-dependent pathway.
Journal of lipid research
Li Y,Yang P,Zhao L,Chen Y,Zhang X,Zeng S,Wei L,Varghese Z,Moorhead JF,Chen Y,Ruan XZ
Mon Apr 01 00:00:00 UTC 2019
SIRT6 promotes the Warburg effect of papillary thyroid cancer cell BCPAP through reactive oxygen species.
OncoTargets and therapy
Yu W,Yang Z,Huang R,Min Z,Ye M
Thu Oct 01 00:00:00 UTC 2020
Protective effects of a neurokinin 1 receptor antagonist on airway epithelial mitochondria dysfunction in asthmatic mice via Nrf2/HO-1 activation.
International immunopharmacology
Wang Z,Li M,Zhou Q,Shang Y
Sun Dec 01 00:00:00 UTC 2019
Involvement of Actin-Regulating Factor Cofilin in the Inclusion Body Formation and RNA Synthesis of Human Parainfluenza Virus Type 3 via Interaction With the Nucleoprotein.
Frontiers in microbiology
Li Y,Zhang C,Lu N,Deng X,Zang G,Zhang S,Tang H,Zhang G
Tue Sep 29 00:00:00 UTC 2020
Hypoxic postconditioning activates the Wnt/β-catenin pathway and protects against transient global cerebral ischemia through Dkk1 Inhibition and GSK-3β inactivation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhan L,Liu D,Wen H,Hu J,Pang T,Sun W,Xu E
Thu Aug 01 00:00:00 UTC 2019
Long non-coding RNA GBCDRlnc1 induces chemoresistance of gallbladder cancer cells by activating autophagy.
Molecular cancer
Cai Q,Wang S,Jin L,Weng M,Zhou D,Wang J,Tang Z,Quan Z
Fri Apr 05 00:00:00 UTC 2019
Inhibition of COX2/PGD2-Related Autophagy Is Involved in the Mechanism of Brain Injury in T2DM Rat.
Frontiers in cellular neuroscience
Yang Y,Chen Q,Zhao Q,Luo Y,Xu Y,Du W,Wang H,Li H,Yang L,Hu C,Zhang J,Li Y,Xia H,Chen Z,Ma J,Tian X,Yang J
Thu Oct 01 00:00:00 UTC 2020
SIRT5 Promotes Hepatocellular Carcinoma Progression by Regulating Mitochondrial Apoptosis.
Journal of Cancer
Zhang R,Wang C,Tian Y,Yao Y,Mao J,Wang H,Li Z,Xu Y,Ye M,Wang L
Thu Oct 01 00:00:00 UTC 2020
iTRAQ-based proteomics analysis reveals the deregulated proteins related to liver toxicity induced by melamine with or without cyanuric acid in mice.
Ecotoxicology and environmental safety
Yin RH,Huang C,Yuan J,Li W,Yin RL,Li HS,Dong Q,Li XT,Bai WL
Sat Jun 15 00:00:00 UTC 2019
DHA-phospholipids (DHA-PL) and EPA-phospholipids (EPA-PL) prevent intestinal dysfunction induced by chronic stress.
Food & function
Cao W,Wang C,Chin Y,Chen X,Gao Y,Yuan S,Xue C,Wang Y,Tang Q
Tue Jan 22 00:00:00 UTC 2019
Enhancing the retrograde axonal transport by curcumin promotes autophagic flux in N2a/APP695swe cells.
Aging
Liang J,Zhou F,Xiong X,Zhang X,Li S,Li X,Gao M,Li Y
Fri Sep 06 00:00:00 UTC 2019
Dexamethasone interferes with osteoblasts formation during osteogenesis through altering IGF-1-mediated angiogenesis.
Journal of cellular physiology
Xu S,Guo R,Li PZ,Li K,Yan Y,Chen J,Wang G,Brand-Saberi B,Yang X,Cheng X
Sun Sep 01 00:00:00 UTC 2019
Role of OXCT1 in ovine adipose and preadipocyte differentiation.
Biochemical and biophysical research communications
Zeng J,Zhou SW,Zhao J,Jin MH,Kang DJ,Yang YX,Wang XL,Chen YL
Tue May 14 00:00:00 UTC 2019
MicroRNA-596 acts as a tumor suppressor in gastric cancer and is upregulated by promotor demethylation.
World journal of gastroenterology
Zhang Z,Dai DQ
Thu Mar 14 00:00:00 UTC 2019
Age-Associated Dopaminergic Neuron Loss and Midbrain Glia Cell Phenotypic Polarization.
Neuroscience
Wang G,Zhou Y,Wang Y,Li D,Liu J,Zhang F
Sun Sep 01 00:00:00 UTC 2019
Fractionated irradiation of right thorax induces abscopal damage on testes leading to decline in fertility.
Scientific reports
Zhang J,Yao D,Song Y,Pan Y,Zhu L,Bai Y,Xu Y,Zhang J,Shao C
Wed Oct 23 00:00:00 UTC 2019
Young Blood Rescues the Cognition of Alzheimer's Model Mice by Restoring the Hippocampal Cholinergic Circuit.
Neuroscience
Xia E,Xu F,Hu C,Kumal JPP,Tang X,Mao D,Li Y,Wu D,Zhang R,Wu S,Sun L
Tue Oct 01 00:00:00 UTC 2019
Hydrogen inhalation protects hypoxic-ischemic brain damage by attenuating inflammation and apoptosis in neonatal rats.
Experimental biology and medicine (Maywood, N.J.)
Wu G,Chen Z,Wang P,Zhao M,Fujino M,Zhang C,Zhou W,Hirano SI,Li XK,Zhao L
Sun Sep 01 00:00:00 UTC 2019
Rutin suppresses FNDC1 expression in bone marrow mesenchymal stem cells to inhibit postmenopausal osteoporosis.
American journal of translational research
Xiao Y,Wei R,Yuan Z,Lan X,Kuang J,Hu D,Song Y,Luo J
Thu Oct 01 00:00:00 UTC 2020
Protective effect of oxytocin on LPS-induced acute lung injury in mice.
Scientific reports
An X,Sun X,Hou Y,Yang X,Chen H,Zhang P,Wu J
Tue Feb 26 00:00:00 UTC 2019
Antinociceptive effect of isoorientin against neuropathic pain induced by the chronic constriction injury of the sciatic nerve in mice.
International immunopharmacology
Zhang G,Liu N,Zhu C,Ma L,Yang J,Du J,Zhang W,Sun T,Niu J,Yu J
Tue Oct 01 00:00:00 UTC 2019
The expression of cerebrospinal fluid exosomal miR-630 plays an important role in the dysfunction of endothelial cells after subarachnoid hemorrhage.
Scientific reports
Sun L,Zhang W,Li Z,Li M,Guo J,Wang H,Wang X
Thu Aug 08 00:00:00 UTC 2019
Alleviation of Hepatic Ischemia Reperfusion Injury by Oleanolic Acid Pretreating via Reducing HMGB1 Release and Inhibiting Apoptosis and Autophagy.
Mediators of inflammation
Wang W,Wu L,Li J,Ji J,Chen K,Yu Q,Li S,Feng J,Liu T,Zhang J,Chen J,Zhou Y,Mao Y,Wang F,Dai W,Fan X,Guo C,Wu J
Mon Feb 24 00:00:00 UTC 2020
Efficient Production of Human Norovirus-Specific IgY in Egg Yolks by Vaccination of Hens with a Recombinant Vesicular Stomatitis Virus Expressing VP1 Protein.
Viruses
Zhu Y,Ma Y,Lu M,Zhang Y,Li A,Liang X,Li J
Thu May 16 00:00:00 UTC 2019
Downregulation of Wnt3 Suppresses Colorectal Cancer Development Through Inhibiting Cell Proliferation and Migration.
Frontiers in pharmacology
Nie X,Xia F,Liu Y,Zhou Y,Ye W,Hean P,Meng J,Liu H,Liu L,Wen J,Ren X,Chen WD,Wang YD
Wed Sep 30 00:00:00 UTC 2020
Normoxia is not favorable for maintaining stemness of human endothelial progenitor cells.
Stem cell research
Lin Y,Liu B,Deng T,Zhong J,Feng Z,Zeng Q,Huang G,Chen Z
Mon Jul 01 00:00:00 UTC 2019
Protective effects of betaine on diabetic induced disruption of the male mice blood-testis barrier by regulating oxidative stress-mediated p38 MAPK pathways.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Jiang YP,Yang JM,Ye RJ,Liu N,Zhang WJ,Ma L,Zheng P,Niu JG,Liu P,Yu JQ
Sun Dec 01 00:00:00 UTC 2019
Exogenous hydrogen sulfide mitigates LPS + ATP-induced inflammation by inhibiting NLRP3 inflammasome activation and promoting autophagy in L02 cells.
Molecular and cellular biochemistry
Wu D,Zhong P,Wang J,Wang H
Mon Jul 01 00:00:00 UTC 2019
Analysis of Toll-Like Receptors in Human Milk: Detection of Membrane-Bound and Soluble Forms.
Journal of immunology research
Cattaneo C,Caramaschi A,Uga E,Braghin M,Cosi G,Peila C,Strozzi MC,Sabatini M,Gazzolo D,Manfredi M,Cavaletto M
Mon May 18 00:00:00 UTC 2020
Mechanisms of Aerobic Exercise Upregulating the Expression of Hippocampal Synaptic Plasticity-Associated Proteins in Diabetic Rats.
Neural plasticity
Li J,Liu Y,Liu B,Li F,Hu J,Wang Q,Li M,Lou S
Mon May 13 00:00:00 UTC 2019
Aspm knockout ferret reveals an evolutionary mechanism governing cerebral cortical size.
Nature
Johnson MB,Sun X,Kodani A,Borges-Monroy R,Girskis KM,Ryu SC,Wang PP,Patel K,Gonzalez DM,Woo YM,Yan Z,Liang B,Smith RS,Chatterjee M,Coman D,Papademetris X,Staib LH,Hyder F,Mandeville JB,Grant PE,Im K,Kwak H,Engelhardt JF,Walsh CA,Bae BI
Sun Apr 01 00:00:00 UTC 2018
Aspm knockout ferret reveals an evolutionary mechanism governing cerebral cortical size.
Nature
Johnson MB,Sun X,Kodani A,Borges-Monroy R,Girskis KM,Ryu SC,Wang PP,Patel K,Gonzalez DM,Woo YM,Yan Z,Liang B,Smith RS,Chatterjee M,Coman D,Papademetris X,Staib LH,Hyder F,Mandeville JB,Grant PE,Im K,Kwak H,Engelhardt JF,Walsh CA,Bae BI
Sun Apr 01 00:00:00 UTC 2018
H2A.Z.1 crosstalk with H3K56-acetylation controls gliogenesis through the transcription of folate receptor.
Nucleic acids research
Su L,Xia W,Shen T,Liang Q,Wang W,Li H,Jiao J
Fri Sep 28 00:00:00 UTC 2018
H2A.Z.1 crosstalk with H3K56-acetylation controls gliogenesis through the transcription of folate receptor.
Nucleic acids research
Su L,Xia W,Shen T,Liang Q,Wang W,Li H,Jiao J
Fri Sep 28 00:00:00 UTC 2018
H2A.Z.1 crosstalk with H3K56-acetylation controls gliogenesis through the transcription of folate receptor.
Nucleic acids research
Su L,Xia W,Shen T,Liang Q,Wang W,Li H,Jiao J
Fri Sep 28 00:00:00 UTC 2018
Decrease of galectin-3 in keratinocytes: A potential diagnostic marker and a critical contributor to the pathogenesis of psoriasis.
Journal of autoimmunity
Shi ZR,Tan GZ,Cao CX,Han YF,Meng Z,Man XY,Jiang ZX,Zhang YP,Dang NN,Wei KH,Bu DF,Liu FT,Wang L
Tue May 01 00:00:00 UTC 2018
Decrease of galectin-3 in keratinocytes: A potential diagnostic marker and a critical contributor to the pathogenesis of psoriasis.
Journal of autoimmunity
Shi ZR,Tan GZ,Cao CX,Han YF,Meng Z,Man XY,Jiang ZX,Zhang YP,Dang NN,Wei KH,Bu DF,Liu FT,Wang L
Tue May 01 00:00:00 UTC 2018
Reversing Multidrug Resistance by Multiplexed Gene Silencing for Enhanced Breast Cancer Chemotherapy.
ACS applied materials & interfaces
Li Y,Gao X,Yu Z,Liu B,Pan W,Li N,Tang B
Wed May 09 00:00:00 UTC 2018
Reversing Multidrug Resistance by Multiplexed Gene Silencing for Enhanced Breast Cancer Chemotherapy.
ACS applied materials & interfaces
Li Y,Gao X,Yu Z,Liu B,Pan W,Li N,Tang B
Wed May 09 00:00:00 UTC 2018
Construction of a High-Efficiency Drug and Gene Co-Delivery System for Cancer Therapy from a pH-Sensitive Supramolecular Inclusion between Oligoethylenimine- graft-β-cyclodextrin and Hyperbranched Polyglycerol Derivative.
ACS applied materials & interfaces
Zhou X,Xu L,Xu J,Wu J,Kirk TB,Ma D,Xue W
Wed Oct 24 00:00:00 UTC 2018
Construction of a High-Efficiency Drug and Gene Co-Delivery System for Cancer Therapy from a pH-Sensitive Supramolecular Inclusion between Oligoethylenimine- graft-β-cyclodextrin and Hyperbranched Polyglycerol Derivative.
ACS applied materials & interfaces
Zhou X,Xu L,Xu J,Wu J,Kirk TB,Ma D,Xue W
Wed Oct 24 00:00:00 UTC 2018
Role of Long Noncoding RNA 799 in the Metastasis of Cervical Cancer through Upregulation of TBL1XR1 Expression.
Molecular therapy. Nucleic acids
Liao LM,Zhang FH,Yao GJ,Ai SF,Zheng M,Huang L
Fri Dec 07 00:00:00 UTC 2018
Role of Long Noncoding RNA 799 in the Metastasis of Cervical Cancer through Upregulation of TBL1XR1 Expression.
Molecular therapy. Nucleic acids
Liao LM,Zhang FH,Yao GJ,Ai SF,Zheng M,Huang L
Fri Dec 07 00:00:00 UTC 2018
Artemisitene suppresses tumorigenesis by inducing DNA damage through deregulating c-Myc-topoisomerase pathway.
Oncogene
Chen J,Li W,Cui K,Ji K,Xu S,Xu Y
Sat Sep 01 00:00:00 UTC 2018
Artemisitene suppresses tumorigenesis by inducing DNA damage through deregulating c-Myc-topoisomerase pathway.
Oncogene
Chen J,Li W,Cui K,Ji K,Xu S,Xu Y
Sat Sep 01 00:00:00 UTC 2018
Cold-induced protein RBM3 orchestrates neurogenesis via modulating Yap mRNA stability in cold stress.
The Journal of cell biology
Xia W,Su L,Jiao J
Mon Oct 01 00:00:00 UTC 2018
Cold-induced protein RBM3 orchestrates neurogenesis via modulating Yap mRNA stability in cold stress.
The Journal of cell biology
Xia W,Su L,Jiao J
Mon Oct 01 00:00:00 UTC 2018
Novel decellularized liver matrix-alginate hybrid gel beads for the 3D culture of hepatocellular carcinoma cells.
International journal of biological macromolecules
Sun D,Liu Y,Wang H,Deng F,Zhang Y,Zhao S,Ma X,Wu H,Sun G
Sun Apr 01 00:00:00 UTC 2018
Novel decellularized liver matrix-alginate hybrid gel beads for the 3D culture of hepatocellular carcinoma cells.
International journal of biological macromolecules
Sun D,Liu Y,Wang H,Deng F,Zhang Y,Zhao S,Ma X,Wu H,Sun G
Sun Apr 01 00:00:00 UTC 2018
Dysfunction of Nrf2-ARE Signaling Pathway: Potential Pathogenesis in the Development of Neurocognitive Impairment in Patients with Moderate to Severe Obstructive Sleep Apnea-Hypopnea Syndrome.
Oxidative medicine and cellular longevity
Zhou L,Ouyang R,Luo H,Peng Y,Chen P,Ren S,Liu G
Wed Dec 19 00:00:00 UTC 2018
Dysfunction of Nrf2-ARE Signaling Pathway: Potential Pathogenesis in the Development of Neurocognitive Impairment in Patients with Moderate to Severe Obstructive Sleep Apnea-Hypopnea Syndrome.
Oxidative medicine and cellular longevity
Zhou L,Ouyang R,Luo H,Peng Y,Chen P,Ren S,Liu G
Wed Dec 19 00:00:00 UTC 2018
Ingredients from Litsea garrettii as Potential Preventive Agents against Oxidative Insult and Inflammatory Response.
Oxidative medicine and cellular longevity
Li YR,Li GH,Sun L,Li L,Liu Y,Kong DG,Wang SQ,Ren DM,Wang XN,Lou HX,Shen T
Mon Oct 22 00:00:00 UTC 2018
Ingredients from Litsea garrettii as Potential Preventive Agents against Oxidative Insult and Inflammatory Response.
Oxidative medicine and cellular longevity
Li YR,Li GH,Sun L,Li L,Liu Y,Kong DG,Wang SQ,Ren DM,Wang XN,Lou HX,Shen T
Mon Oct 22 00:00:00 UTC 2018
Bexarotene protects against neurotoxicity partially through a PPARγ-dependent mechanism in mice following traumatic brain injury.
Neurobiology of disease
He J,Liu H,Zhong J,Guo Z,Wu J,Zhang H,Huang Z,Jiang L,Li H,Zhang Z,Liu L,Wu Y,Qi L,Sun X,Cheng C
Sat Sep 01 00:00:00 UTC 2018
Bexarotene protects against neurotoxicity partially through a PPARγ-dependent mechanism in mice following traumatic brain injury.
Neurobiology of disease
He J,Liu H,Zhong J,Guo Z,Wu J,Zhang H,Huang Z,Jiang L,Li H,Zhang Z,Liu L,Wu Y,Qi L,Sun X,Cheng C
Sat Sep 01 00:00:00 UTC 2018
Nitric oxide, afferent sensory nerves, and antioxidative enzymes in the mechanism of protection mediated by tricarbonyldichlororuthenium(II) dimer and sodium hydrosulfide against aspirin-induced gastric damage.
Journal of gastroenterology
Magierowski M,Hubalewska-Mazgaj M,Magierowska K,Wojcik D,Sliwowski Z,Kwiecien S,Brzozowski T
Mon Jan 01 00:00:00 UTC 2018
Nitric oxide, afferent sensory nerves, and antioxidative enzymes in the mechanism of protection mediated by tricarbonyldichlororuthenium(II) dimer and sodium hydrosulfide against aspirin-induced gastric damage.
Journal of gastroenterology
Magierowski M,Hubalewska-Mazgaj M,Magierowska K,Wojcik D,Sliwowski Z,Kwiecien S,Brzozowski T
Mon Jan 01 00:00:00 UTC 2018
PD-L1 confers glioblastoma multiforme malignancy via Ras binding and Ras/Erk/EMT activation.
Biochimica et biophysica acta. Molecular basis of disease
Qiu XY,Hu DX,Chen WQ,Chen RQ,Qian SR,Li CY,Li YJ,Xiong XX,Liu D,Pan F,Yu SB,Chen XQ
Tue May 01 00:00:00 UTC 2018
PD-L1 confers glioblastoma multiforme malignancy via Ras binding and Ras/Erk/EMT activation.
Biochimica et biophysica acta. Molecular basis of disease
Qiu XY,Hu DX,Chen WQ,Chen RQ,Qian SR,Li CY,Li YJ,Xiong XX,Liu D,Pan F,Yu SB,Chen XQ
Tue May 01 00:00:00 UTC 2018
The inhibitory effect in Fraxinellone on oxidative stress-induced senescence correlates with AMP-activated protein kinase-dependent autophagy restoration.
Journal of cellular physiology
Han X,Chen H,Zhou J,Tai H,Gong H,Wang X,Huang N,Qin J,Fang T,Wang F,Xiao H
Tue May 01 00:00:00 UTC 2018
The inhibitory effect in Fraxinellone on oxidative stress-induced senescence correlates with AMP-activated protein kinase-dependent autophagy restoration.
Journal of cellular physiology
Han X,Chen H,Zhou J,Tai H,Gong H,Wang X,Huang N,Qin J,Fang T,Wang F,Xiao H
Tue May 01 00:00:00 UTC 2018
Anticancer effect of SZC015 on pancreatic cancer via mitochondria-dependent apoptosis and the constitutive suppression of activated nuclear factor κB and STAT3 in vitro and in vivo.
Journal of cellular physiology
Song Y,Gao L,Tang Z,Li H,Sun B,Chu P,Qaed E,Ma X,Peng J,Wang S,Hu M,Tang Z
Mon Jan 01 00:00:00 UTC 2018
Anticancer effect of SZC015 on pancreatic cancer via mitochondria-dependent apoptosis and the constitutive suppression of activated nuclear factor κB and STAT3 in vitro and in vivo.
Journal of cellular physiology
Song Y,Gao L,Tang Z,Li H,Sun B,Chu P,Qaed E,Ma X,Peng J,Wang S,Hu M,Tang Z
Mon Jan 01 00:00:00 UTC 2018
Lysyl oxidase blockade ameliorates anovulation in polycystic ovary syndrome.
Human reproduction (Oxford, England)
Zhang C,Ma J,Wang W,Sun Y,Sun K
Thu Nov 01 00:00:00 UTC 2018
Lysyl oxidase blockade ameliorates anovulation in polycystic ovary syndrome.
Human reproduction (Oxford, England)
Zhang C,Ma J,Wang W,Sun Y,Sun K
Thu Nov 01 00:00:00 UTC 2018
Combination Treatment of Icariin and L-DOPA Against 6-OHDA-Lesioned Dopamine Neurotoxicity.
Frontiers in molecular neuroscience
Lu DS,Chen C,Zheng YX,Li DD,Wang GQ,Liu J,Shi J,Zhang F
Thu Oct 01 00:00:00 UTC 2020
Combination Treatment of Icariin and L-DOPA Against 6-OHDA-Lesioned Dopamine Neurotoxicity.
Frontiers in molecular neuroscience
Lu DS,Chen C,Zheng YX,Li DD,Wang GQ,Liu J,Shi J,Zhang F
Thu Oct 01 00:00:00 UTC 2020
Regulation on SIRT1-PGC-1α/Nrf2 pathway together with selective inhibition of aldose reductase makes compound hr5F a potential agent for the treatment of diabetic complications.
Biochemical pharmacology
Wang Z,Yuan S,Li Y,Zhang Z,Xiao W,Tang D,Ye K,Liu Z,Wang C,Zheng Y,Nie H,Chen H
Sun Apr 01 00:00:00 UTC 2018
Regulation on SIRT1-PGC-1α/Nrf2 pathway together with selective inhibition of aldose reductase makes compound hr5F a potential agent for the treatment of diabetic complications.
Biochemical pharmacology
Wang Z,Yuan S,Li Y,Zhang Z,Xiao W,Tang D,Ye K,Liu Z,Wang C,Zheng Y,Nie H,Chen H
Sun Apr 01 00:00:00 UTC 2018
Exogenous oxidants activate nuclear factor kappa B through Toll-like receptor 4 stimulation to maintain inflammatory phenotype in macrophage.
Biochemical pharmacology
Zhang Y,Igwe OJ
Mon Jan 01 00:00:00 UTC 2018
Exogenous oxidants activate nuclear factor kappa B through Toll-like receptor 4 stimulation to maintain inflammatory phenotype in macrophage.
Biochemical pharmacology
Zhang Y,Igwe OJ
Mon Jan 01 00:00:00 UTC 2018
Glutathione Reduction of Patulin-Evoked Cytotoxicity in HEK293 Cells by the Prevention of Oxidative Damage and the Mitochondrial Apoptotic Pathway.
Journal of agricultural and food chemistry
Wang X,Jin C,Zhong Y,Li X,Han J,Xue W,Wu P,Xia X,Peng X
Wed Jul 25 00:00:00 UTC 2018
Glutathione Reduction of Patulin-Evoked Cytotoxicity in HEK293 Cells by the Prevention of Oxidative Damage and the Mitochondrial Apoptotic Pathway.
Journal of agricultural and food chemistry
Wang X,Jin C,Zhong Y,Li X,Han J,Xue W,Wu P,Xia X,Peng X
Wed Jul 25 00:00:00 UTC 2018
Curcumin attenuates resistance to irinotecan via induction of apoptosis of cancer stem cells in chemoresistant colon cancer cells.
International journal of oncology
Su P,Yang Y,Wang G,Chen X,Ju Y
Sat Sep 01 00:00:00 UTC 2018
Curcumin attenuates resistance to irinotecan via induction of apoptosis of cancer stem cells in chemoresistant colon cancer cells.
International journal of oncology
Su P,Yang Y,Wang G,Chen X,Ju Y
Sat Sep 01 00:00:00 UTC 2018
Protective effects of ginsenoside Rg1 against lipopolysaccharide/d-galactosamine-induced acute liver injury in mice through inhibiting toll-like receptor 4 signaling pathway.
International immunopharmacology
Ning C,Gao X,Wang C,Huo X,Liu Z,Sun H,Yang X,Sun P,Ma X,Meng Q,Liu K
Wed Aug 01 00:00:00 UTC 2018
Protective effects of ginsenoside Rg1 against lipopolysaccharide/d-galactosamine-induced acute liver injury in mice through inhibiting toll-like receptor 4 signaling pathway.
International immunopharmacology
Ning C,Gao X,Wang C,Huo X,Liu Z,Sun H,Yang X,Sun P,Ma X,Meng Q,Liu K
Wed Aug 01 00:00:00 UTC 2018
Activation of Nrf2 pathway and inhibition of NLRP3 inflammasome activation contribute to the protective effect of chlorogenic acid on acute liver injury.
International immunopharmacology
Shi A,Shi H,Wang Y,Liu X,Cheng Y,Li H,Zhao H,Wang S,Dong L
Mon Jan 01 00:00:00 UTC 2018
Activation of Nrf2 pathway and inhibition of NLRP3 inflammasome activation contribute to the protective effect of chlorogenic acid on acute liver injury.
International immunopharmacology
Shi A,Shi H,Wang Y,Liu X,Cheng Y,Li H,Zhao H,Wang S,Dong L
Mon Jan 01 00:00:00 UTC 2018
Down-regulation of IFIT3 protects liver from ischemia-reperfusion injury.
International immunopharmacology
Guan G,Shen Y,Yu Q,Liu H,Zhang B,Guo Y,Zhu X,Li Z,Rao W,Zhuang L,Zang Y
Sun Jul 01 00:00:00 UTC 2018
Down-regulation of IFIT3 protects liver from ischemia-reperfusion injury.
International immunopharmacology
Guan G,Shen Y,Yu Q,Liu H,Zhang B,Guo Y,Zhu X,Li Z,Rao W,Zhuang L,Zang Y
Sun Jul 01 00:00:00 UTC 2018
Oxyresveratrol prevents lipopolysaccharide/d-galactosamine-induced acute liver injury in mice.
International immunopharmacology
Jia YN,Lu HP,Peng YL,Zhang BS,Gong XB,Su J,Zhou Y,Pan MH,Xu L
Thu Mar 01 00:00:00 UTC 2018
Oxyresveratrol prevents lipopolysaccharide/d-galactosamine-induced acute liver injury in mice.
International immunopharmacology
Jia YN,Lu HP,Peng YL,Zhang BS,Gong XB,Su J,Zhou Y,Pan MH,Xu L
Thu Mar 01 00:00:00 UTC 2018
BF211, a derivative of bufalin, enhances the cytocidal effects in multiple myeloma cells by inhibiting the IL-6/JAK2/STAT3 pathway.
International immunopharmacology
Wu XY,Tian F,Su MH,Wu M,Huang Y,Hu LH,Jin L,Zhu XJ
Thu Nov 01 00:00:00 UTC 2018
BF211, a derivative of bufalin, enhances the cytocidal effects in multiple myeloma cells by inhibiting the IL-6/JAK2/STAT3 pathway.
International immunopharmacology
Wu XY,Tian F,Su MH,Wu M,Huang Y,Hu LH,Jin L,Zhu XJ
Thu Nov 01 00:00:00 UTC 2018
Leucine Carboxyl Methyltransferase Downregulation and Protein Phosphatase Methylesterase Upregulation Contribute Toward the Inhibition of Protein Phosphatase 2A by α-Synuclein.
Frontiers in aging neuroscience
Tian H,Lu Y,Liu J,Liu W,Lu L,Duan C,Gao G,Yang H
Tue Sep 29 00:00:00 UTC 2020
Leucine Carboxyl Methyltransferase Downregulation and Protein Phosphatase Methylesterase Upregulation Contribute Toward the Inhibition of Protein Phosphatase 2A by α-Synuclein.
Frontiers in aging neuroscience
Tian H,Lu Y,Liu J,Liu W,Lu L,Duan C,Gao G,Yang H
Tue Sep 29 00:00:00 UTC 2020
Cdk5 Inhibitory Peptide Prevents Loss of Dopaminergic Neurons and Alleviates Behavioral Changes in an MPTP Induced Parkinson's Disease Mouse Model.
Frontiers in aging neuroscience
He R,Huang W,Huang Y,Xu M,Song P,Huang Y,Xie H,Hu Y
Wed Sep 30 00:00:00 UTC 2020
Cdk5 Inhibitory Peptide Prevents Loss of Dopaminergic Neurons and Alleviates Behavioral Changes in an MPTP Induced Parkinson's Disease Mouse Model.
Frontiers in aging neuroscience
He R,Huang W,Huang Y,Xu M,Song P,Huang Y,Xie H,Hu Y
Wed Sep 30 00:00:00 UTC 2020
Baicalein inhibits cervical cancer progression via downregulating long noncoding RNA BDLNR and its downstream PI3K/Akt pathway.
The international journal of biochemistry & cell biology
Yu X,Yang Y,Li Y,Cao Y,Tang L,Chen F,Xia J
Mon Jan 01 00:00:00 UTC 2018
Baicalein inhibits cervical cancer progression via downregulating long noncoding RNA BDLNR and its downstream PI3K/Akt pathway.
The international journal of biochemistry & cell biology
Yu X,Yang Y,Li Y,Cao Y,Tang L,Chen F,Xia J
Mon Jan 01 00:00:00 UTC 2018
Baicalein inhibits cervical cancer progression via downregulating long noncoding RNA BDLNR and its downstream PI3K/Akt pathway.
The international journal of biochemistry & cell biology
Yu X,Yang Y,Li Y,Cao Y,Tang L,Chen F,Xia J
Mon Jan 01 00:00:00 UTC 2018
CHOP induces apoptosis by affecting brain iron metabolism in rats with subarachnoid hemorrhage.
Experimental neurology
Zhao J,Xiang X,Zhang H,Jiang D,Liang Y,Qing W,Liu L,Zhao Q,He Z
Sun Apr 01 00:00:00 UTC 2018
CHOP induces apoptosis by affecting brain iron metabolism in rats with subarachnoid hemorrhage.
Experimental neurology
Zhao J,Xiang X,Zhang H,Jiang D,Liang Y,Qing W,Liu L,Zhao Q,He Z
Sun Apr 01 00:00:00 UTC 2018
2',4'-Dihydroxy-6'-methoxy-3',5'-dimethylchalcone, a potent Nrf2/ARE pathway inhibitor, reverses drug resistance by decreasing glutathione synthesis and drug efflux in BEL-7402/5-FU cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Wei X,Mo X,An F,Ji X,Lu Y
Sat Sep 01 00:00:00 UTC 2018
2',4'-Dihydroxy-6'-methoxy-3',5'-dimethylchalcone, a potent Nrf2/ARE pathway inhibitor, reverses drug resistance by decreasing glutathione synthesis and drug efflux in BEL-7402/5-FU cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Wei X,Mo X,An F,Ji X,Lu Y
Sat Sep 01 00:00:00 UTC 2018
Intermittent streptozotocin administration induces behavioral and pathological features relevant to Alzheimer's disease and vascular dementia.
Neuropharmacology
Murtishaw AS,Heaney CF,Bolton MM,Belmonte KCD,Langhardt MA,Kinney JW
Sun Jul 15 00:00:00 UTC 2018
Intermittent streptozotocin administration induces behavioral and pathological features relevant to Alzheimer's disease and vascular dementia.
Neuropharmacology
Murtishaw AS,Heaney CF,Bolton MM,Belmonte KCD,Langhardt MA,Kinney JW
Sun Jul 15 00:00:00 UTC 2018
Anti-inflammatory activities and potential mechanisms of phenolic acids isolated from Salvia miltiorrhiza f. alba roots in THP-1 macrophages.
Journal of ethnopharmacology
Liu H,Ma S,Xia H,Lou H,Zhu F,Sun L
Fri Aug 10 00:00:00 UTC 2018
Anti-inflammatory activities and potential mechanisms of phenolic acids isolated from Salvia miltiorrhiza f. alba roots in THP-1 macrophages.
Journal of ethnopharmacology
Liu H,Ma S,Xia H,Lou H,Zhu F,Sun L
Fri Aug 10 00:00:00 UTC 2018
Di-(2-ethylhexyl)-phthalate induces glucose metabolic disorder in adolescent rats.
Environmental science and pollution research international
Xu J,Zhou L,Wang S,Zhu J,Liu T,Jia Y,Sun D,Chen H,Wang Q,Xu F,Zhang Y,Liu H,Zhang T,Ye L
Thu Feb 01 00:00:00 UTC 2018
Di-(2-ethylhexyl)-phthalate induces glucose metabolic disorder in adolescent rats.
Environmental science and pollution research international
Xu J,Zhou L,Wang S,Zhu J,Liu T,Jia Y,Sun D,Chen H,Wang Q,Xu F,Zhang Y,Liu H,Zhang T,Ye L
Thu Feb 01 00:00:00 UTC 2018
Whole-exome sequencing revealed HKDC1 as a candidate gene associated with autosomal-recessive retinitis pigmentosa.
Human molecular genetics
Zhang L,Sun Z,Zhao P,Huang L,Xu M,Yang Y,Chen X,Lu F,Zhang X,Wang H,Zhang S,Liu W,Jiang Z,Ma S,Chen R,Zhao C,Yang Z,Sui R,Zhu X
Sat Dec 01 00:00:00 UTC 2018
Dietary eicosapentaenoic acid normalizes hippocampal omega-3 and 6 polyunsaturated fatty acid profile, attenuates glial activation and regulates BDNF function in a rodent model of neuroinflammation induced by central interleukin-1β administration.
European journal of nutrition
Dong Y,Xu M,Kalueff AV,Song C
Wed Aug 01 00:00:00 UTC 2018
Dietary eicosapentaenoic acid normalizes hippocampal omega-3 and 6 polyunsaturated fatty acid profile, attenuates glial activation and regulates BDNF function in a rodent model of neuroinflammation induced by central interleukin-1β administration.
European journal of nutrition
Dong Y,Xu M,Kalueff AV,Song C
Wed Aug 01 00:00:00 UTC 2018
CNTNAP4 Impacts Epilepsy Through GABAA Receptors Regulation: Evidence From Temporal Lobe Epilepsy Patients and Mouse Models.
Cerebral cortex (New York, N.Y. : 1991)
Shangguan Y,Xu X,Ganbat B,Li Y,Wang W,Yang Y,Lu X,Du C,Tian X,Wang X
Mon Oct 01 00:00:00 UTC 2018
CNTNAP4 Impacts Epilepsy Through GABAA Receptors Regulation: Evidence From Temporal Lobe Epilepsy Patients and Mouse Models.
Cerebral cortex (New York, N.Y. : 1991)
Shangguan Y,Xu X,Ganbat B,Li Y,Wang W,Yang Y,Lu X,Du C,Tian X,Wang X
Mon Oct 01 00:00:00 UTC 2018
MiR-20a-3p regulates TGF-β1/Survivin pathway to affect keratinocytes proliferation and apoptosis by targeting SFMBT1 in vitro.
Cellular signalling
Li R,Qiao M,Zhao X,Yan J,Wang X,Sun Q
Sat Sep 01 00:00:00 UTC 2018
MiR-20a-3p regulates TGF-β1/Survivin pathway to affect keratinocytes proliferation and apoptosis by targeting SFMBT1 in vitro.
Cellular signalling
Li R,Qiao M,Zhao X,Yan J,Wang X,Sun Q
Sat Sep 01 00:00:00 UTC 2018
Erythropoietin Treatment Ameliorates Lupus Nephritis of MRL/lpr Mice.
Inflammation
Zhang Z,Liu D,Zhang X,Wang X
Mon Oct 01 00:00:00 UTC 2018
Erythropoietin Treatment Ameliorates Lupus Nephritis of MRL/lpr Mice.
Inflammation
Zhang Z,Liu D,Zhang X,Wang X
Mon Oct 01 00:00:00 UTC 2018
Serum and Ectopic Endometrium from Women with Endometriosis Modulate Macrophage M1/M2 Polarization via the Smad2/Smad3 Pathway.
Journal of immunology research
Nie MF,Xie Q,Wu YH,He H,Zou LJ,She XL,Wu XQ
Thu Dec 27 00:00:00 UTC 2018
Serum and Ectopic Endometrium from Women with Endometriosis Modulate Macrophage M1/M2 Polarization via the Smad2/Smad3 Pathway.
Journal of immunology research
Nie MF,Xie Q,Wu YH,He H,Zou LJ,She XL,Wu XQ
Thu Dec 27 00:00:00 UTC 2018
FGF9 modulates Schwann cell myelination in developing nerves and induces a pro-inflammatory environment during injury.
Journal of cellular biochemistry
Deng B,Lv W,Duan W,Liu Y,Li Z,Song X,Cui C,Qi X,Wang X,Li C
Thu Nov 01 00:00:00 UTC 2018
FGF9 modulates Schwann cell myelination in developing nerves and induces a pro-inflammatory environment during injury.
Journal of cellular biochemistry
Deng B,Lv W,Duan W,Liu Y,Li Z,Song X,Cui C,Qi X,Wang X,Li C
Thu Nov 01 00:00:00 UTC 2018
The N-terminal ubiquitin-associated domain of Cezanne is crucial for its function to suppress NF-κB pathway.
Journal of cellular biochemistry
Ji Y,Cao L,Zeng L,Zhang Z,Xiao Q,Guan P,Chen S,Chen Y,Wang M,Guo D
Thu Feb 01 00:00:00 UTC 2018
The N-terminal ubiquitin-associated domain of Cezanne is crucial for its function to suppress NF-κB pathway.
Journal of cellular biochemistry
Ji Y,Cao L,Zeng L,Zhang Z,Xiao Q,Guan P,Chen S,Chen Y,Wang M,Guo D
Thu Feb 01 00:00:00 UTC 2018
High Expression of lncRNA AFAP1-AS1 Promotes the Progression of Colon Cancer and Predicts Poor Prognosis.
Journal of Cancer
Bo H,Fan L,Li J,Liu Z,Zhang S,Shi L,Guo C,Li X,Liao Q,Zhang W,Zhou M,Xiang B,Li X,Li G,Xiong W,Zeng Z,Xiong F,Gong Z
Thu Oct 01 00:00:00 UTC 2020
High Expression of lncRNA AFAP1-AS1 Promotes the Progression of Colon Cancer and Predicts Poor Prognosis.
Journal of Cancer
Bo H,Fan L,Li J,Liu Z,Zhang S,Shi L,Guo C,Li X,Liao Q,Zhang W,Zhou M,Xiang B,Li X,Li G,Xiong W,Zeng Z,Xiong F,Gong Z
Thu Oct 01 00:00:00 UTC 2020
Neuroprotective Effect of Anethole Against Neuropathic Pain Induced by Chronic Constriction Injury of the Sciatic Nerve in Mice.
Neurochemical research
Wang B,Zhang G,Yang M,Liu N,Li YX,Ma H,Ma L,Sun T,Tan H,Yu J
Sat Dec 01 00:00:00 UTC 2018
Neuroprotective Effect of Anethole Against Neuropathic Pain Induced by Chronic Constriction Injury of the Sciatic Nerve in Mice.
Neurochemical research
Wang B,Zhang G,Yang M,Liu N,Li YX,Ma H,Ma L,Sun T,Tan H,Yu J
Sat Dec 01 00:00:00 UTC 2018
All-Trans Retinoic Acid Ameliorates the Early Experimental Cerebral Ischemia-Reperfusion Injury in Rats by Inhibiting the Loss of the Blood-Brain Barrier via the JNK/P38MAPK Signaling Pathway.
Neurochemical research
Li M,Tian X,An R,Yang M,Zhang Q,Xiang F,Liu H,Wang Y,Xu L,Dong Z
Fri Jun 01 00:00:00 UTC 2018
All-Trans Retinoic Acid Ameliorates the Early Experimental Cerebral Ischemia-Reperfusion Injury in Rats by Inhibiting the Loss of the Blood-Brain Barrier via the JNK/P38MAPK Signaling Pathway.
Neurochemical research
Li M,Tian X,An R,Yang M,Zhang Q,Xiang F,Liu H,Wang Y,Xu L,Dong Z
Fri Jun 01 00:00:00 UTC 2018
Long non-coding RNA (lncRNA) MAGI2-AS3 inhibits breast cancer cell growth by targeting the Fas/FasL signalling pathway.
Human cell
Yang Y,Yang H,Xu M,Zhang H,Sun M,Mu P,Dong T,Du S,Liu K
Sun Jul 01 00:00:00 UTC 2018
Long non-coding RNA (lncRNA) MAGI2-AS3 inhibits breast cancer cell growth by targeting the Fas/FasL signalling pathway.
Human cell
Yang Y,Yang H,Xu M,Zhang H,Sun M,Mu P,Dong T,Du S,Liu K
Sun Jul 01 00:00:00 UTC 2018
Effects of calycosin against high-fat diet-induced nonalcoholic fatty liver disease in mice.
Journal of gastroenterology and hepatology
Duan X,Meng Q,Wang C,Liu Z,Sun H,Huo X,Sun P,Ma X,Peng J,Liu K
Thu Feb 01 00:00:00 UTC 2018
Effects of calycosin against high-fat diet-induced nonalcoholic fatty liver disease in mice.
Journal of gastroenterology and hepatology
Duan X,Meng Q,Wang C,Liu Z,Sun H,Huo X,Sun P,Ma X,Peng J,Liu K
Thu Feb 01 00:00:00 UTC 2018
Natural antisense transcript of hypoxia-inducible factor 1 regulates hypoxic cell apoptosis in epithelial ovarian cancer.
OncoTargets and therapy
Qiu JJ,Lin XJ,Zheng TT,Tang XY,Hua KQ
Wed Mar 30 00:00:00 UTC 2022
Natural antisense transcript of hypoxia-inducible factor 1 regulates hypoxic cell apoptosis in epithelial ovarian cancer.
OncoTargets and therapy
Qiu JJ,Lin XJ,Zheng TT,Tang XY,Hua KQ
Wed Mar 30 00:00:00 UTC 2022
Contribution of dysregulated circRNA_100876 to proliferation and metastasis of esophageal squamous cell carcinoma.
OncoTargets and therapy
Cao S,Chen G,Yan L,Li L,Huang X
Sun Apr 10 00:00:00 UTC 2022
Contribution of dysregulated circRNA_100876 to proliferation and metastasis of esophageal squamous cell carcinoma.
OncoTargets and therapy
Cao S,Chen G,Yan L,Li L,Huang X
Sun Apr 10 00:00:00 UTC 2022
Wnt/β-catenin modulates chronic tobacco smoke exposure-induced acquisition of pulmonary cancer stem cell properties and diallyl trisulfide intervention.
Toxicology letters
Wang J,Chen J,Jiang Y,Shi Y,Zhu J,Xie C,Geng S,Wu J,Zhang Q,Wang X,Meng Y,Li Y,Chen Y,Cao W,Wang X,Zhong C,Li X
Sun Jul 01 00:00:00 UTC 2018
Wnt/β-catenin modulates chronic tobacco smoke exposure-induced acquisition of pulmonary cancer stem cell properties and diallyl trisulfide intervention.
Toxicology letters
Wang J,Chen J,Jiang Y,Shi Y,Zhu J,Xie C,Geng S,Wu J,Zhang Q,Wang X,Meng Y,Li Y,Chen Y,Cao W,Wang X,Zhong C,Li X
Sun Jul 01 00:00:00 UTC 2018
Association between Tim‑3 and Gal‑9 expression and gastric cancer prognosis.
Oncology reports
Wang Y,Zhao E,Zhang Z,Zhao G,Cao H
Mon Oct 01 00:00:00 UTC 2018
Association between Tim‑3 and Gal‑9 expression and gastric cancer prognosis.
Oncology reports
Wang Y,Zhao E,Zhang Z,Zhao G,Cao H
Mon Oct 01 00:00:00 UTC 2018
Snx27 Deletion Promotes Recovery From Spinal Cord Injury by Neuroprotection and Reduces Macrophage/Microglia Proliferation.
Frontiers in neurology
Zeng Y,Wang N,Guo T,Zheng Q,Wang S,Wu S,Li X,Wu J,Chen Z,Xu H,Wang X,Lin B
Thu Oct 01 00:00:00 UTC 2020
Snx27 Deletion Promotes Recovery From Spinal Cord Injury by Neuroprotection and Reduces Macrophage/Microglia Proliferation.
Frontiers in neurology
Zeng Y,Wang N,Guo T,Zheng Q,Wang S,Wu S,Li X,Wu J,Chen Z,Xu H,Wang X,Lin B
Thu Oct 01 00:00:00 UTC 2020
Lithium chloride inhibits cell survival, overcomes drug resistance, and triggers apoptosis in multiple myeloma via activation of the Wnt/β-catenin pathway.
American journal of translational research
Yao R,Sun X,Xie Y,Liu L,Han D,Yao Y,Li H,Li Z,Xu K
Wed Sep 30 00:00:00 UTC 2020
Lithium chloride inhibits cell survival, overcomes drug resistance, and triggers apoptosis in multiple myeloma via activation of the Wnt/β-catenin pathway.
American journal of translational research
Yao R,Sun X,Xie Y,Liu L,Han D,Yao Y,Li H,Li Z,Xu K
Wed Sep 30 00:00:00 UTC 2020
Acrolein-induced autophagy-dependent apoptosis via activation of the lysosomal-mitochondrial pathway in EAhy926 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Luo S,Jiang L,Li Q,Sun X,Liu T,Pei F,Zhang T,Liu T,Dong L,Liu X,Jiang L
Mon Oct 01 00:00:00 UTC 2018
Acrolein-induced autophagy-dependent apoptosis via activation of the lysosomal-mitochondrial pathway in EAhy926 cells.
Toxicology in vitro : an international journal published in association with BIBRA
Luo S,Jiang L,Li Q,Sun X,Liu T,Pei F,Zhang T,Liu T,Dong L,Liu X,Jiang L
Mon Oct 01 00:00:00 UTC 2018
Relationship of Epidermal Growth Factor Receptor Expression with Clinical Symptoms and Metastasis of Invasive Breast Cancer.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Han L,Li L,Wang N,Xiong Y,Li Y,Gu Y
Sat Dec 01 00:00:00 UTC 2018
Relationship of Epidermal Growth Factor Receptor Expression with Clinical Symptoms and Metastasis of Invasive Breast Cancer.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Han L,Li L,Wang N,Xiong Y,Li Y,Gu Y
Sat Dec 01 00:00:00 UTC 2018
Resveratrol relieves particulate matter (mean diameter < 2.5 μm)-induced oxidative injury of lung cells through attenuation of autophagy deregulation.
Journal of applied toxicology : JAT
Li Y,Qian W,Wang D,Meng Y,Wang X,Chen Y,Li X,Xie C,Zhong C,Fu S
Sat Sep 01 00:00:00 UTC 2018
Resveratrol relieves particulate matter (mean diameter < 2.5 μm)-induced oxidative injury of lung cells through attenuation of autophagy deregulation.
Journal of applied toxicology : JAT
Li Y,Qian W,Wang D,Meng Y,Wang X,Chen Y,Li X,Xie C,Zhong C,Fu S
Sat Sep 01 00:00:00 UTC 2018
Formononetin inhibits colon carcinoma cell growth and invasion by microRNA‑149‑mediated EphB3 downregulation and inhibition of PI3K/AKT and STAT3 signaling pathways.
Molecular medicine reports
Wang AL,Li Y,Zhao Q,Fan LQ
Fri Jun 01 00:00:00 UTC 2018
Formononetin inhibits colon carcinoma cell growth and invasion by microRNA‑149‑mediated EphB3 downregulation and inhibition of PI3K/AKT and STAT3 signaling pathways.
Molecular medicine reports
Wang AL,Li Y,Zhao Q,Fan LQ
Fri Jun 01 00:00:00 UTC 2018
Construction and protective immunogenicity of DNA vaccine pNMB0315 against Neisseria meningitidis serogroup B.
Molecular medicine reports
Wu X,Li K,Xie M,Yu M,Tang S,Li Z,Hu S
Thu Feb 01 00:00:00 UTC 2018
Construction and protective immunogenicity of DNA vaccine pNMB0315 against Neisseria meningitidis serogroup B.
Molecular medicine reports
Wu X,Li K,Xie M,Yu M,Tang S,Li Z,Hu S
Thu Feb 01 00:00:00 UTC 2018
Pyroptosis of HUVECs can be induced by heat stroke.
Biochemical and biophysical research communications
Pei Y,Geng Y,Su L
Fri Nov 30 00:00:00 UTC 2018
Pyroptosis of HUVECs can be induced by heat stroke.
Biochemical and biophysical research communications
Pei Y,Geng Y,Su L
Fri Nov 30 00:00:00 UTC 2018
The protective role of low-concentration alcohol in high-fructose induced adverse cardiovascular events in mice.
Biochemical and biophysical research communications
Wu X,Pan B,Wang Y,Liu L,Huang X,Tian J
Mon Jan 01 00:00:00 UTC 2018
The protective role of low-concentration alcohol in high-fructose induced adverse cardiovascular events in mice.
Biochemical and biophysical research communications
Wu X,Pan B,Wang Y,Liu L,Huang X,Tian J
Mon Jan 01 00:00:00 UTC 2018
Chronic cold exposure results in subcutaneous adipose tissue browning and altered global metabolism in Qinghai-Tibetan plateau pika (Ochotona curzoniae).
Biochemical and biophysical research communications
Li J,Yang Q,Bai Z,Zhou W,Semenza GL,Ge RL
Sat Jun 02 00:00:00 UTC 2018
Chronic cold exposure results in subcutaneous adipose tissue browning and altered global metabolism in Qinghai-Tibetan plateau pika (Ochotona curzoniae).
Biochemical and biophysical research communications
Li J,Yang Q,Bai Z,Zhou W,Semenza GL,Ge RL
Sat Jun 02 00:00:00 UTC 2018
AMPK alleviates endoplasmic reticulum stress by inducing the ER-chaperone ORP150 via FOXO1 to protect human bronchial cells from apoptosis.
Biochemical and biophysical research communications
Liu JQ,Zhang L,Yao J,Yao S,Yuan T
Sun Mar 04 00:00:00 UTC 2018
AMPK alleviates endoplasmic reticulum stress by inducing the ER-chaperone ORP150 via FOXO1 to protect human bronchial cells from apoptosis.
Biochemical and biophysical research communications
Liu JQ,Zhang L,Yao J,Yao S,Yuan T
Sun Mar 04 00:00:00 UTC 2018
ERα is a negative regulator of PD-L1 gene transcription in breast cancer.
Biochemical and biophysical research communications
Liu L,Shen Y,Zhu X,Lv R,Li S,Zhang Z,Shi YG,Tan L
Sat Oct 20 00:00:00 UTC 2018
ERα is a negative regulator of PD-L1 gene transcription in breast cancer.
Biochemical and biophysical research communications
Liu L,Shen Y,Zhu X,Lv R,Li S,Zhang Z,Shi YG,Tan L
Sat Oct 20 00:00:00 UTC 2018
The Potential Protective Effect of Curcumin on Amyloid-β-42 Induced Cytotoxicity in HT-22 Cells.
BioMed research international
Zhang L,Fang Y,Cheng X,Lian Y,Zeng Z,Wu C,Zhu H,Xu H
Thu Sep 06 00:00:00 UTC 2018
The Potential Protective Effect of Curcumin on Amyloid-β-42 Induced Cytotoxicity in HT-22 Cells.
BioMed research international
Zhang L,Fang Y,Cheng X,Lian Y,Zeng Z,Wu C,Zhu H,Xu H
Thu Sep 06 00:00:00 UTC 2018
DL-3-n-butylphthalide alleviates vascular cognitive impairment induced by chronic cerebral hypoperfusion by activating the Akt/Nrf2 signaling pathway in the hippocampus of rats.
Neuroscience letters
Qi Q,Xu J,Lv P,Dong Y,Liu Z,Hu M,Xiao Y,Jia Y,Jin W,Fan M,Zhang D,Meng N
Fri Apr 13 00:00:00 UTC 2018
DL-3-n-butylphthalide alleviates vascular cognitive impairment induced by chronic cerebral hypoperfusion by activating the Akt/Nrf2 signaling pathway in the hippocampus of rats.
Neuroscience letters
Qi Q,Xu J,Lv P,Dong Y,Liu Z,Hu M,Xiao Y,Jia Y,Jin W,Fan M,Zhang D,Meng N
Fri Apr 13 00:00:00 UTC 2018
Expression of tumor suppressor programmed cell death 4 in endometrioid endometrial carcinomas and clinicopathological significance.
Oncology letters
Liu Y,Sun H,Mao H,Gao M,Tan X,Li Y,Li Y,Muloye GM,Zhang L,Wang X,Wei Z
Fri Jun 01 00:00:00 UTC 2018
Expression of tumor suppressor programmed cell death 4 in endometrioid endometrial carcinomas and clinicopathological significance.
Oncology letters
Liu Y,Sun H,Mao H,Gao M,Tan X,Li Y,Li Y,Muloye GM,Zhang L,Wang X,Wei Z
Fri Jun 01 00:00:00 UTC 2018
Overexpression of SMARCA2 or CAMK2D is associated with cisplatin resistance in human epithelial ovarian cancer.
Oncology letters
Xu X,Zheng Z,Jia L,Suo S,Liu B,Shao T,Tu Q,Hua Y,Xu H
Sat Sep 01 00:00:00 UTC 2018
Overexpression of SMARCA2 or CAMK2D is associated with cisplatin resistance in human epithelial ovarian cancer.
Oncology letters
Xu X,Zheng Z,Jia L,Suo S,Liu B,Shao T,Tu Q,Hua Y,Xu H
Sat Sep 01 00:00:00 UTC 2018
TAM Infiltration Differences in "Tumor-First" and " ZHENG-First" Models and the Underlying Inflammatory Molecular Mechanism in Pancreatic Cancer.
Integrative cancer therapies
Wang FJ,Wang P,Chen LY,Geng YW,Chen H,Meng ZQ,Liu LM,Chen Z
Sat Sep 01 00:00:00 UTC 2018
TAM Infiltration Differences in "Tumor-First" and " ZHENG-First" Models and the Underlying Inflammatory Molecular Mechanism in Pancreatic Cancer.
Integrative cancer therapies
Wang FJ,Wang P,Chen LY,Geng YW,Chen H,Meng ZQ,Liu LM,Chen Z
Sat Sep 01 00:00:00 UTC 2018
Lipoxin A4 improves erectile dysfunction in rats with type I diabetes by inhibiting oxidative stress and corporal fibrosis.
Asian journal of andrology
Cui K,Tang Z,Li CC,Wang T,Rao K,Wang SG,Liu JH,Chen Z
Tue May 14 00:00:00 UTC 2019
Lipoxin A4 improves erectile dysfunction in rats with type I diabetes by inhibiting oxidative stress and corporal fibrosis.
Asian journal of andrology
Cui K,Tang Z,Li CC,Wang T,Rao K,Wang SG,Liu JH,Chen Z
Tue May 14 00:00:00 UTC 2019
Nicotine Induces Progressive Properties of Lung Adenocarcinoma A549 Cells by Inhibiting Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) Expression and Plasma Membrane Localization.
Technology in cancer research & treatment
Li H,Ma N,Wang J,Wang Y,Yuan C,Wu J,Luo M,Yang J,Chen J,Shi J,Liu X
Mon Jan 01 00:00:00 UTC 2018
Nicotine Induces Progressive Properties of Lung Adenocarcinoma A549 Cells by Inhibiting Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) Expression and Plasma Membrane Localization.
Technology in cancer research & treatment
Li H,Ma N,Wang J,Wang Y,Yuan C,Wu J,Luo M,Yang J,Chen J,Shi J,Liu X
Mon Jan 01 00:00:00 UTC 2018
Expression of diaphragmatic myostatin and correlation with apoptosis in rats with chronic obstructive pulmonary disease.
Experimental and therapeutic medicine
Zhou X,Yi D,Wu Y,Pei X,Yu H,Chen Y,Jiang Y,Li W
Thu Mar 01 00:00:00 UTC 2018
Expression of diaphragmatic myostatin and correlation with apoptosis in rats with chronic obstructive pulmonary disease.
Experimental and therapeutic medicine
Zhou X,Yi D,Wu Y,Pei X,Yu H,Chen Y,Jiang Y,Li W
Thu Mar 01 00:00:00 UTC 2018
Effects of SB202190 on expression levels of IL-6 and NF-κB in flap ischemia-reperfusion injury.
Experimental and therapeutic medicine
Guo K,Ma J,Liang W
Sat Sep 01 00:00:00 UTC 2018
Effects of SB202190 on expression levels of IL-6 and NF-κB in flap ischemia-reperfusion injury.
Experimental and therapeutic medicine
Guo K,Ma J,Liang W
Sat Sep 01 00:00:00 UTC 2018
Donor Treatment With a Hypoxia-Inducible Factor-1 Agonist Prevents Donation After Cardiac Death Liver Graft Injury in a Rat Isolated Perfusion Model.
Artificial organs
Zhang X,Liu Z,Xiao Q,Zeng C,Lai CH,Fan X,Ye Q,Wang Y,Xiong Y
Thu Mar 01 00:00:00 UTC 2018
Donor Treatment With a Hypoxia-Inducible Factor-1 Agonist Prevents Donation After Cardiac Death Liver Graft Injury in a Rat Isolated Perfusion Model.
Artificial organs
Zhang X,Liu Z,Xiao Q,Zeng C,Lai CH,Fan X,Ye Q,Wang Y,Xiong Y
Thu Mar 01 00:00:00 UTC 2018
Effects of Chicory on Serum Uric Acid, Renal Function, and GLUT9 Expression in Hyperuricaemic Rats with Renal Injury and In Vitro Verification with Cells.
Evidence-based complementary and alternative medicine : eCAM
Jin YN,Lin ZJ,Zhang B,Bai YF
Mon Apr 25 00:00:00 UTC 2022
Effects of Chicory on Serum Uric Acid, Renal Function, and GLUT9 Expression in Hyperuricaemic Rats with Renal Injury and In Vitro Verification with Cells.
Evidence-based complementary and alternative medicine : eCAM
Jin YN,Lin ZJ,Zhang B,Bai YF
Mon Apr 25 00:00:00 UTC 2022
Low-Dose Radiation Enhanced Inhibition of Breast Tumor Xenograft and Reduced Myocardial Injury Induced by Doxorubicin.
Dose-response : a publication of International Hormesis Society
Jiang X,Du Y,Meng X,Zhang H,Zhao D,Zhao L,Chen J,Xiao S,Jiang H
Thu Oct 01 00:00:00 UTC 2020
Low-Dose Radiation Enhanced Inhibition of Breast Tumor Xenograft and Reduced Myocardial Injury Induced by Doxorubicin.
Dose-response : a publication of International Hormesis Society
Jiang X,Du Y,Meng X,Zhang H,Zhao D,Zhao L,Chen J,Xiao S,Jiang H
Thu Oct 01 00:00:00 UTC 2020
Fucosterol Protects against Concanavalin A-Induced Acute Liver Injury: Focus on P38 MAPK/NF-κB Pathway Activity.
Gastroenterology research and practice
Mo W,Wang C,Li J,Chen K,Xia Y,Li S,Xu L,Lu X,Wang W,Guo C
Fri Mar 18 00:00:00 UTC 2022
Fucosterol Protects against Concanavalin A-Induced Acute Liver Injury: Focus on P38 MAPK/NF-κB Pathway Activity.
Gastroenterology research and practice
Mo W,Wang C,Li J,Chen K,Xia Y,Li S,Xu L,Lu X,Wang W,Guo C
Fri Mar 18 00:00:00 UTC 2022
Proteomics analysis of altered proteins in kidney of mice with aristolochic acid nephropathy using the fluorogenic derivatization-liquid chromatography-tandem mass spectrometry method.
Biomedical chromatography : BMC
Lin CE,Chang WS,Lee JA,Chang TY,Huang YS,Hirasaki Y,Chen HS,Imai K,Chen SM
Thu Mar 01 00:00:00 UTC 2018
Proteomics analysis of altered proteins in kidney of mice with aristolochic acid nephropathy using the fluorogenic derivatization-liquid chromatography-tandem mass spectrometry method.
Biomedical chromatography : BMC
Lin CE,Chang WS,Lee JA,Chang TY,Huang YS,Hirasaki Y,Chen HS,Imai K,Chen SM
Thu Mar 01 00:00:00 UTC 2018
Arsenic sulfide amplifies JQ1 toxicity via mitochondrial pathway in gastric and colon cancer cells.
Drug design, development and therapy
Tan Z,Zhang X,Kang T,Zhang L,Chen S
Tue Mar 26 00:00:00 UTC 2019
Arsenic sulfide amplifies JQ1 toxicity via mitochondrial pathway in gastric and colon cancer cells.
Drug design, development and therapy
Tan Z,Zhang X,Kang T,Zhang L,Chen S
Tue Mar 26 00:00:00 UTC 2019
miR-383 negatively regulates osteoblastic differentiation of bone marrow mesenchymal stem cells in rats by targeting Satb2.
Bone
Tang J,Zhang Z,Jin X,Shi H
Sat Sep 01 00:00:00 UTC 2018
miR-383 negatively regulates osteoblastic differentiation of bone marrow mesenchymal stem cells in rats by targeting Satb2.
Bone
Tang J,Zhang Z,Jin X,Shi H
Sat Sep 01 00:00:00 UTC 2018
High Concentration of Aspirin Induces Apoptosis in Rat Tendon Stem Cells via Inhibition of the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Y,Tang H,He G,Shi Y,Kang X,Lyu J,Zhou M,Zhu M,Zhang J,Tang K
Tue Dec 11 00:00:00 UTC 2018
High Concentration of Aspirin Induces Apoptosis in Rat Tendon Stem Cells via Inhibition of the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Y,Tang H,He G,Shi Y,Kang X,Lyu J,Zhou M,Zhu M,Zhang J,Tang K
Tue Dec 11 00:00:00 UTC 2018
Mesenchymal Stem Cell-Derived Exosomes Reduce A1 Astrocytes via Downregulation of Phosphorylated NFκB P65 Subunit in Spinal Cord Injury.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang L,Pei S,Han L,Guo B,Li Y,Duan R,Yao Y,Xue B,Chen X,Jia Y
Fri Nov 09 00:00:00 UTC 2018
Mesenchymal Stem Cell-Derived Exosomes Reduce A1 Astrocytes via Downregulation of Phosphorylated NFκB P65 Subunit in Spinal Cord Injury.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang L,Pei S,Han L,Guo B,Li Y,Duan R,Yao Y,Xue B,Chen X,Jia Y
Fri Nov 09 00:00:00 UTC 2018
The FOXK1-CCDC43 Axis Promotes the Invasion and Metastasis of Colorectal Cancer Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang J,Liu G,Liu M,Xiang L,Xiao Y,Zhu H,Wu X,Peng Y,Zhang W,Jiang P,Li A,Nan Q,Chen Y,Chen C,Cheng T,Liu S,Wang J
Tue Jan 29 00:00:00 UTC 2019
The FOXK1-CCDC43 Axis Promotes the Invasion and Metastasis of Colorectal Cancer Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang J,Liu G,Liu M,Xiang L,Xiao Y,Zhu H,Wu X,Peng Y,Zhang W,Jiang P,Li A,Nan Q,Chen Y,Chen C,Cheng T,Liu S,Wang J
Tue Jan 29 00:00:00 UTC 2019
Tumor-Secreted Exosomal miR-222 Promotes Tumor Progression via Regulating P27 Expression and Re-Localization in Pancreatic Cancer.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li Z,Tao Y,Wang X,Jiang P,Li J,Peng M,Zhang X,Chen K,Liu H,Zhen P,Zhu J,Liu X,Liu X
Tue Dec 11 00:00:00 UTC 2018
Tumor-Secreted Exosomal miR-222 Promotes Tumor Progression via Regulating P27 Expression and Re-Localization in Pancreatic Cancer.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li Z,Tao Y,Wang X,Jiang P,Li J,Peng M,Zhang X,Chen K,Liu H,Zhen P,Zhu J,Liu X,Liu X
Tue Dec 11 00:00:00 UTC 2018
Cancer-derived exosomal miR-25-3p promotes pre-metastatic niche formation by inducing vascular permeability and angiogenesis.
Nature communications
Zeng Z,Li Y,Pan Y,Lan X,Song F,Sun J,Zhou K,Liu X,Ren X,Wang F,Hu J,Zhu X,Yang W,Liao W,Li G,Ding Y,Liang L
Wed Dec 19 00:00:00 UTC 2018
Cancer-derived exosomal miR-25-3p promotes pre-metastatic niche formation by inducing vascular permeability and angiogenesis.
Nature communications
Zeng Z,Li Y,Pan Y,Lan X,Song F,Sun J,Zhou K,Liu X,Ren X,Wang F,Hu J,Zhu X,Yang W,Liao W,Li G,Ding Y,Liang L
Wed Dec 19 00:00:00 UTC 2018
An E-selectin targeting and MMP-2-responsive dextran-curcumin polymeric prodrug for targeted therapy of acute kidney injury.
Biomaterials science
Hu JB,Liu D,Qi J,Lu KJ,Jin FY,Ying XY,You J,Du YZ
Tue Nov 20 00:00:00 UTC 2018
Interfering RNA against PKC-α inhibits TNF-α-induced IP3R1 expression and improves glomerular filtration rate in rats with fulminant hepatic failure.
American journal of physiology. Renal physiology
Wang DL,Dai WY,Wang W,Wen Y,Zhou Y,Zhao YT,Wu J,Liu P
Tue May 01 00:00:00 UTC 2018
TRPC1/3/6 inhibition attenuates the TGF-β1-induced epithelial-mesenchymal transition in gastric cancer via the Ras/Raf1/ERK signaling pathway.
Cell biology international
Ge P,Wei L,Zhang M,Hu B,Wang K,Li Y,Liu S,Wang J,Li Y
Wed Aug 01 00:00:00 UTC 2018
lncRNA GAS5 inhibits colorectal cancer cell proliferation via the miR‑182‑5p/FOXO3a axis.
Oncology reports
Cheng K,Zhao Z,Wang G,Wang J,Zhu W
Mon Oct 01 00:00:00 UTC 2018
Knockdown of long noncoding RNA GHET1 inhibits cell‑cycle progression and invasion of gastric cancer cells.
Molecular medicine reports
Xia Y,Yan Z,Wan Y,Wei S,Bi Y,Zhao J,Liu J,Liao DJ,Huang H
Sat Sep 01 00:00:00 UTC 2018
lncRNA DLEU1 contributes to tumorigenesis and development of endometrial carcinoma by targeting mTOR.
Molecular carcinogenesis
Du Y,Wang L,Chen S,Liu Y,Zhao Y
Sat Sep 01 00:00:00 UTC 2018
WISP1 mediates lung injury following hepatic ischemia reperfusion dependent on TLR4 in mice.
BMC pulmonary medicine
Tong Y,Yu Z,Zhang R,Ding X,Chen Z,Li Q
Thu Dec 06 00:00:00 UTC 2018
WISP1 mediates lung injury following hepatic ischemia reperfusion dependent on TLR4 in mice.
BMC pulmonary medicine
Tong Y,Yu Z,Zhang R,Ding X,Chen Z,Li Q
Thu Dec 06 00:00:00 UTC 2018
Hepatic, Metabolic, and Toxicity Evaluation of Repeated Oral Administration of SnS2 Nanoflowers in Mice.
Toxicological sciences : an official journal of the Society of Toxicology
Bai D,Li Q,Xiong Y,Wang C,Shen P,Bai L,Yuan L,Wu P
Wed Aug 01 00:00:00 UTC 2018
Hepatic, Metabolic, and Toxicity Evaluation of Repeated Oral Administration of SnS2 Nanoflowers in Mice.
Toxicological sciences : an official journal of the Society of Toxicology
Bai D,Li Q,Xiong Y,Wang C,Shen P,Bai L,Yuan L,Wu P
Wed Aug 01 00:00:00 UTC 2018
Oxymatrine Inhibits Transforming Growth Factor β1 (TGF-β1)-Induced Cardiac Fibroblast-to-Myofibroblast Transformation (FMT) by Mediating the Notch Signaling Pathway In Vitro.
Medical science monitor : international medical journal of experimental and clinical research
Zhao L,Xu Y,Tao L,Yang Y,Shen X,Li L,Luo P
Sun Sep 09 00:00:00 UTC 2018
Oxymatrine Inhibits Transforming Growth Factor β1 (TGF-β1)-Induced Cardiac Fibroblast-to-Myofibroblast Transformation (FMT) by Mediating the Notch Signaling Pathway In Vitro.
Medical science monitor : international medical journal of experimental and clinical research
Zhao L,Xu Y,Tao L,Yang Y,Shen X,Li L,Luo P
Sun Sep 09 00:00:00 UTC 2018
Experimental evidence of good efficacy and reduced toxicity with peptide-doxorubicin to treat gastric cancer.
Oncotarget
Zhang J,Yuan JP,Wang Q,Shao LH,Liu SP,Firestone RA,Hong YP,Li JG,Xin YC,Li Y
Fri Jan 05 00:00:00 UTC 2018
Experimental evidence of good efficacy and reduced toxicity with peptide-doxorubicin to treat gastric cancer.
Oncotarget
Zhang J,Yuan JP,Wang Q,Shao LH,Liu SP,Firestone RA,Hong YP,Li JG,Xin YC,Li Y
Fri Jan 05 00:00:00 UTC 2018
Early Postnatal Exposure to Airborne Fine Particulate Matter Induces Autism-like Phenotypes in Male Rats.
Toxicological sciences : an official journal of the Society of Toxicology
Li K,Li L,Cui B,Gai Z,Li Q,Wang S,Yan J,Lin B,Tian L,Liu H,Liu X,Xi Z
Thu Mar 01 00:00:00 UTC 2018
Early Postnatal Exposure to Airborne Fine Particulate Matter Induces Autism-like Phenotypes in Male Rats.
Toxicological sciences : an official journal of the Society of Toxicology
Li K,Li L,Cui B,Gai Z,Li Q,Wang S,Yan J,Lin B,Tian L,Liu H,Liu X,Xi Z
Thu Mar 01 00:00:00 UTC 2018
In vitro inhibition of porcine reproductive and respiratory syndrome virus replication by short antisense oligonucleotides with locked nucleic acid modification.
BMC veterinary research
Zhu L,Bi J,Zheng L,Zhao Q,Shu X,Guo G,Liu J,Yang G,Liu J,Yin G
Mon Mar 26 00:00:00 UTC 2018
In vitro inhibition of porcine reproductive and respiratory syndrome virus replication by short antisense oligonucleotides with locked nucleic acid modification.
BMC veterinary research
Zhu L,Bi J,Zheng L,Zhao Q,Shu X,Guo G,Liu J,Yang G,Liu J,Yin G
Mon Mar 26 00:00:00 UTC 2018
Luteolin sensitizes the antitumor effect of cisplatin in drug-resistant ovarian cancer via induction of apoptosis and inhibition of cell migration and invasion.
Journal of ovarian research
Wang H,Luo Y,Qiao T,Wu Z,Huang Z
Mon Nov 19 00:00:00 UTC 2018
Luteolin sensitizes the antitumor effect of cisplatin in drug-resistant ovarian cancer via induction of apoptosis and inhibition of cell migration and invasion.
Journal of ovarian research
Wang H,Luo Y,Qiao T,Wu Z,Huang Z
Mon Nov 19 00:00:00 UTC 2018
FPOA induces apoptosis in HeLa human cervical cancer cells through a caspase-mediated pathway.
Oncology letters
Li X,Bau T,Bao H
Fri Jun 01 00:00:00 UTC 2018
Crotonols A and B, two rare tigliane diterpenoid derivatives against K562 cells from Croton tiglium.
Organic & biomolecular chemistry
Wang J,Qin L,Zhao B,Cai L,Zhong Z,Liu Y,Zhou X
Wed Dec 19 00:00:00 UTC 2018
Pseudo-Ginsenoside Rh2 induces A549 cells apoptosis via the Ras/Raf/ERK/p53 pathway.
Experimental and therapeutic medicine
Wang Y,Xu H,Lu Z,Yu X,Lv C,Tian Y,Sui D
Fri Jun 01 00:00:00 UTC 2018
Thrombopoietin protects H9C2 cells from excessive autophagy and apoptosis in doxorubicin-induced cardiotoxicity.
Oncology letters
Wang H,Wang H,Liang EY,Zhou LX,Dong ZL,Liang P,Weng QF,Yang M
Mon Jan 01 00:00:00 UTC 2018
High expression of WNT7A predicts poor prognosis and promote tumor metastasis in pancreatic ductal adenocarcinoma.
Scientific reports
Wu DJ,Jiang YS,He RZ,Tao LY,Yang MW,Fu XL,Yang JY,Zhu K
Thu Oct 25 00:00:00 UTC 2018
High expression of WNT7A predicts poor prognosis and promote tumor metastasis in pancreatic ductal adenocarcinoma.
Scientific reports
Wu DJ,Jiang YS,He RZ,Tao LY,Yang MW,Fu XL,Yang JY,Zhu K
Thu Oct 25 00:00:00 UTC 2018
Effects of Tumor Necrosis Factor α (TNF-α) and Interleukina 10 (IL-10) on Intercellular Cell Adhesion Molecule-1 (ICAM-1) and Cluster of Differentiation 31 (CD31) in Human Coronary Artery Endothelial Cells.
Medical science monitor : international medical journal of experimental and clinical research
Xue M,Qiqige C,Zhang Q,Zhao H,Su L,Sun P,Zhao P
Wed Jun 27 00:00:00 UTC 2018
Effects of Tumor Necrosis Factor α (TNF-α) and Interleukina 10 (IL-10) on Intercellular Cell Adhesion Molecule-1 (ICAM-1) and Cluster of Differentiation 31 (CD31) in Human Coronary Artery Endothelial Cells.
Medical science monitor : international medical journal of experimental and clinical research
Xue M,Qiqige C,Zhang Q,Zhao H,Su L,Sun P,Zhao P
Wed Jun 27 00:00:00 UTC 2018
Trio-Based Deep Sequencing Reveals a Low Incidence of Off-Target Mutations in the Offspring of Genetically Edited Goats.
Frontiers in genetics
Li C,Zhou S,Li Y,Li G,Ding Y,Li L,Liu J,Qu L,Sonstegard T,Huang X,Jiang Y,Chen Y,Petersen B,Wang X
Tue Sep 29 00:00:00 UTC 2020
Trio-Based Deep Sequencing Reveals a Low Incidence of Off-Target Mutations in the Offspring of Genetically Edited Goats.
Frontiers in genetics
Li C,Zhou S,Li Y,Li G,Ding Y,Li L,Liu J,Qu L,Sonstegard T,Huang X,Jiang Y,Chen Y,Petersen B,Wang X
Tue Sep 29 00:00:00 UTC 2020
LncRNA PCGEM1 Induces Ovarian Carcinoma Tumorigenesis and Progression Through RhoA Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Chen S,Wang LL,Sun KX,Liu Y,Guan X,Zong ZH,Zhao Y
Wed Aug 15 00:00:00 UTC 2018
LncRNA PCGEM1 Induces Ovarian Carcinoma Tumorigenesis and Progression Through RhoA Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Chen S,Wang LL,Sun KX,Liu Y,Guan X,Zong ZH,Zhao Y
Wed Aug 15 00:00:00 UTC 2018
RACK1 is indispensable for porcine reproductive and respiratory syndrome virus replication and NF-κB activation in Marc-145 cells.
Scientific reports
Bi J,Zhao Q,Zhu L,Li X,Yang G,Liu J,Yin G
Wed Feb 14 00:00:00 UTC 2018
Targeting MAPK Pathways by Naringenin Modulates Microglia M1/M2 Polarization in Lipopolysaccharide-Stimulated Cultures.
Frontiers in cellular neuroscience
Zhang B,Wei YZ,Wang GQ,Li DD,Shi JS,Zhang F
Thu Oct 01 00:00:00 UTC 2020
Cationic nanomicelles derived from Pluronic F127 as delivery vehicles of Chinese herbal medicine active components of ursolic acid for colorectal cancer treatment.
RSC advances
Yan Z,Wang Q,Liu X,Peng J,Li Q,Wu M,Lin J
Fri Apr 27 00:00:00 UTC 2018
Retraction: Cd induces G2/M cell cycle arrest by up-regulating miR-133b via directly targeting PPP2R2D in L02 hepatocytes.
Metallomics : integrated biometal science
Zhang Y,Xiao F,Zhong C,Zeng M,Zou L
Wed Nov 14 00:00:00 UTC 2018
miR-429 suppresses tumor migration and invasion by targeting CRKL in hepatocellular carcinoma via inhibiting Raf/MEK/ERK pathway and epithelial-mesenchymal transition.
Scientific reports
Guo C,Zhao D,Zhang Q,Liu S,Sun MZ
Mon Feb 05 00:00:00 UTC 2018
Metformin suppresses growth and adrenocorticotrophic hormone secretion in mouse pituitary corticotroph tumor AtT20 cells.
Molecular and cellular endocrinology
Jin K,Ruan L,Pu J,Zhong A,Wang F,Tan S,Huang H,Mu J,Yang G
Sat Dec 15 00:00:00 UTC 2018
Fangchinoline Ameliorates the Expressions of Angiogenic Molecule in Cerebral Ischemia Induced Neuronal Degeneration in Neonatal Rats.
Translational neuroscience
Daicheng H,Shiwen X,Huaping Z,Yong L,Qianqian Z,Changxia H
Thu Mar 31 00:00:00 UTC 2022
Genipin alleviates high-fat diet-induced hyperlipidemia and hepatic lipid accumulation in mice via miR-142a-5p/SREBP-1c axis.
The FEBS journal
Zhong H,Chen K,Feng M,Shao W,Wu J,Chen K,Liang T,Liu C
Thu Feb 01 00:00:00 UTC 2018
Collagen-based three-dimensional culture microenvironment promotes epithelial to mesenchymal transition and drug resistance of human ovarian cancer in vitro.
RSC advances
Liu M,Zhang X,Long C,Xu H,Cheng X,Chang J,Zhang C,Zhang C,Wang X
Fri Feb 23 00:00:00 UTC 2018
Necrostatin-1 promotes ectopic periodontal tissue like structure regeneration in LPS-treated PDLSCs.
PloS one
Yan B,Zhang H,Dai T,Gu Y,Qiu X,Hu C,Liu Y,Wei K,Li D
Fri Apr 19 00:00:00 UTC 2019
Necrostatin-1 promotes ectopic periodontal tissue like structure regeneration in LPS-treated PDLSCs.
PloS one
Yan B,Zhang H,Dai T,Gu Y,Qiu X,Hu C,Liu Y,Wei K,Li D
Fri Apr 19 00:00:00 UTC 2019
The Traditional Chinese Medicine MLC901 inhibits inflammation processes after focal cerebral ischemia.
Scientific reports
Widmann C,Gandin C,Petit-Paitel A,Lazdunski M,Heurteaux C
Mon Dec 24 00:00:00 UTC 2018
The Traditional Chinese Medicine MLC901 inhibits inflammation processes after focal cerebral ischemia.
Scientific reports
Widmann C,Gandin C,Petit-Paitel A,Lazdunski M,Heurteaux C
Mon Dec 24 00:00:00 UTC 2018
Lactobacillus helveticus KLDS1.8701 alleviates d-galactose-induced aging by regulating Nrf-2 and gut microbiota in mice.
Food & function
Li B,Evivie SE,Lu J,Jiao Y,Wang C,Li Z,Liu F,Huo G
Thu Dec 13 00:00:00 UTC 2018
Phosphoglycerate Mutase 1 (PGAM1) Promotes Pancreatic Ductal Adenocarcinoma (PDAC) Metastasis by Acting as a Novel Downstream Target of the PI3K/Akt/mTOR Pathway.
Oncology research
Liu X,Tan X,Liu P,Wu Y,Qian S,Zhang X
Thu Aug 23 00:00:00 UTC 2018
Adipocytes fuel gastric cancer omental metastasis via PITPNC1-mediated fatty acid metabolic reprogramming.
Theranostics
Tan Y,Lin K,Zhao Y,Wu Q,Chen D,Wang J,Liang Y,Li J,Hu J,Wang H,Liu Y,Zhang S,He W,Huang Q,Hu X,Yao Z,Liang B,Liao W,Shi M
Fri Sep 06 00:00:00 UTC 2019
Adipocytes fuel gastric cancer omental metastasis via PITPNC1-mediated fatty acid metabolic reprogramming.
Theranostics
Tan Y,Lin K,Zhao Y,Wu Q,Chen D,Wang J,Liang Y,Li J,Hu J,Wang H,Liu Y,Zhang S,He W,Huang Q,Hu X,Yao Z,Liang B,Liao W,Shi M
Fri Sep 06 00:00:00 UTC 2019
Transcription Factor FOXO3a Is a Negative Regulator of Cytotoxicity of Fusarium mycotoxin in GES-1 Cells.
Toxicological sciences : an official journal of the Society of Toxicology
Yang Y,Yu S,Liu N,Xu H,Gong Y,Wu Y,Wang P,Su X,Liao Y,De Saeger S,Humpf HU,Wu A
Sat Dec 01 00:00:00 UTC 2018
Transcription Factor FOXO3a Is a Negative Regulator of Cytotoxicity of Fusarium mycotoxin in GES-1 Cells.
Toxicological sciences : an official journal of the Society of Toxicology
Yang Y,Yu S,Liu N,Xu H,Gong Y,Wu Y,Wang P,Su X,Liao Y,De Saeger S,Humpf HU,Wu A
Sat Dec 01 00:00:00 UTC 2018
MicroRNA‑106a regulates the proliferation and invasion of human osteosarcoma cells by targeting VNN2.
Oncology reports
Chen Y,Huang T,Yang X,Liu C,Li P,Wang Z,Zhi S
Mon Oct 01 00:00:00 UTC 2018
7-Acetylsinumaximol B Induces Apoptosis and Autophagy in Human Gastric Carcinoma Cells through Mitochondria Dysfunction and Activation of the PERK/eIF2α/ATF4/CHOP Signaling Pathway.
Marine drugs
Tsai TC,Lai KH,Su JH,Wu YJ,Sheu JH
Mon Mar 26 00:00:00 UTC 2018
7-Acetylsinumaximol B Induces Apoptosis and Autophagy in Human Gastric Carcinoma Cells through Mitochondria Dysfunction and Activation of the PERK/eIF2α/ATF4/CHOP Signaling Pathway.
Marine drugs
Tsai TC,Lai KH,Su JH,Wu YJ,Sheu JH
Mon Mar 26 00:00:00 UTC 2018
Marine Bacterial Polysaccharide EPS11 Inhibits Cancer Cell Growth via Blocking Cell Adhesion and Stimulating Anoikis.
Marine drugs
Cao R,Jin W,Shan Y,Wang J,Liu G,Kuang S,Sun C
Thu Mar 08 00:00:00 UTC 2018
Marine Bacterial Polysaccharide EPS11 Inhibits Cancer Cell Growth via Blocking Cell Adhesion and Stimulating Anoikis.
Marine drugs
Cao R,Jin W,Shan Y,Wang J,Liu G,Kuang S,Sun C
Thu Mar 08 00:00:00 UTC 2018
Knockdown of TIPE2 increases the proliferation in lipopolysaccharide-stimulated gastric cancer cells.
BMC cancer
Liu W,Fan Y,Shi Y,Lin Z,Huang X,Huang W,Shen D,Qi Z
Wed Aug 29 00:00:00 UTC 2018
Knockdown of TIPE2 increases the proliferation in lipopolysaccharide-stimulated gastric cancer cells.
BMC cancer
Liu W,Fan Y,Shi Y,Lin Z,Huang X,Huang W,Shen D,Qi Z
Wed Aug 29 00:00:00 UTC 2018
Long Noncoding RNA HOTTIP Promotes Mouse Hepatic Stellate Cell Activation via Downregulating miR-148a.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li Z,Wang J,Zeng Q,Hu C,Zhang J,Wang H,Yan J,Li H,Yu Z
Tue Jan 29 00:00:00 UTC 2019
Long Noncoding RNA HOTTIP Promotes Mouse Hepatic Stellate Cell Activation via Downregulating miR-148a.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li Z,Wang J,Zeng Q,Hu C,Zhang J,Wang H,Yan J,Li H,Yu Z
Tue Jan 29 00:00:00 UTC 2019
The influence of GAPT extraction on synapse loss of APPswe/PS1dE9 transgenic mice via adjusting Bcl-2/Bax balance.
Alzheimer's & dementia (New York, N. Y.)
Shi J,Zhang X,Ni J,Wei M,Li T,Zhou B,Liu X,Zhang L,Wang P,Tian J,Wang Y
Wed Mar 30 00:00:00 UTC 2022
The influence of GAPT extraction on synapse loss of APPswe/PS1dE9 transgenic mice via adjusting Bcl-2/Bax balance.
Alzheimer's & dementia (New York, N. Y.)
Shi J,Zhang X,Ni J,Wei M,Li T,Zhou B,Liu X,Zhang L,Wang P,Tian J,Wang Y
Wed Mar 30 00:00:00 UTC 2022
Th17 cell-derived IL-17A promoted tumor progression via STAT3/NF-κB/Notch1 signaling in non-small cell lung cancer.
Oncoimmunology
Wang R,Yang L,Zhang C,Wang R,Zhang Z,He Q,Chen X,Zhang B,Qin Z,Wang L,Zhang Y
Thu Feb 04 00:00:00 UTC 2021
miR‑92b promotes autophagy and suppresses viability and invasion in breast cancer by targeting EZH2.
International journal of oncology
Liu F,Sang M,Meng L,Gu L,Liu S,Li J,Geng C
Mon Oct 01 00:00:00 UTC 2018
Atrazine exposure improves the proliferation of H22 cells in vitro and in vivo.
RSC advances
Tian Y,He J,Liu N,Huang D,Liu Z,Yang Y,Chen J,Zhao B,Zhao S,Liang B
Wed Jun 13 00:00:00 UTC 2018
Temozolomide-Perillyl alcohol conjugate impairs Mitophagy flux by inducing lysosomal dysfunction in non-small cell lung Cancer cells and sensitizes them to irradiation.
Journal of experimental & clinical cancer research : CR
Chang M,Song X,Geng X,Wang X,Wang W,Chen TC,Xie L,Song X
Tue Oct 16 00:00:00 UTC 2018
Temozolomide-Perillyl alcohol conjugate impairs Mitophagy flux by inducing lysosomal dysfunction in non-small cell lung Cancer cells and sensitizes them to irradiation.
Journal of experimental & clinical cancer research : CR
Chang M,Song X,Geng X,Wang X,Wang W,Chen TC,Xie L,Song X
Tue Oct 16 00:00:00 UTC 2018
Polybrominated Diphenyl Ethers Quinone Induced Parthanatos-like Cell Death through a Reactive Oxygen Species-Associated Poly(ADP-ribose) Polymerase 1 Signaling.
Chemical research in toxicology
Dong W,Yang B,Wang Y,Yuan J,Fan Y,Song E,Song Y
Mon Nov 19 00:00:00 UTC 2018
RING finger protein 10 prevents neointimal hyperplasia by promoting apoptosis in vitro and in vivo.
Life sciences
Yu G,Chen J,Li S,Pu P,Huang W,Zhao Y,Peng X,Wang R,Lei H
Sat Sep 01 00:00:00 UTC 2018
[Impact of Cystic Fibrosis Transmembrane Conductance Regulator on Malignant Properties of KRAS Mutant Lung Adenocarcinoma A549 Cells].
Zhongguo fei ai za zhi = Chinese journal of lung cancer
Li H,Wang Y,Yang J,Liu X,Shi J
Tue Feb 20 00:00:00 UTC 2018
Epidermal Growth Factor, through Alleviating Oxidative Stress, Protect IPEC-J2 Cells from Lipopolysaccharides-Induced Apoptosis.
International journal of molecular sciences
Tang X,Liu B,Wang X,Yu Q,Fang R
Wed Mar 14 00:00:00 UTC 2018
Epidermal Growth Factor, through Alleviating Oxidative Stress, Protect IPEC-J2 Cells from Lipopolysaccharides-Induced Apoptosis.
International journal of molecular sciences
Tang X,Liu B,Wang X,Yu Q,Fang R
Wed Mar 14 00:00:00 UTC 2018
Regional up-regulation of NOX2 contributes to the differential vulnerability of outer hair cells to neomycin.
Biochemical and biophysical research communications
Qi M,Qiu Y,Zhou X,Tian K,Zhou K,Sun F,Yue B,Chen F,Zha D,Qiu J
Sat Jun 02 00:00:00 UTC 2018
Lactate enhanced the effect of parathyroid hormone on osteoblast differentiation via GPR81-PKC-Akt signaling.
Biochemical and biophysical research communications
Wu Y,Wang M,Zhang K,Li Y,Xu M,Tang S,Qu X,Li C
Wed Sep 05 00:00:00 UTC 2018
Histone variant H3.3 orchestrates neural stem cell differentiation in the developing brain.
Cell death and differentiation
Xia W,Jiao J
Fri Sep 01 00:00:00 UTC 2017
Histone variant H3.3 orchestrates neural stem cell differentiation in the developing brain.
Cell death and differentiation
Xia W,Jiao J
Fri Sep 01 00:00:00 UTC 2017
PLAA Mutations Cause a Lethal Infantile Epileptic Encephalopathy by Disrupting Ubiquitin-Mediated Endolysosomal Degradation of Synaptic Proteins.
American journal of human genetics
Hall EA,Nahorski MS,Murray LM,Shaheen R,Perkins E,Dissanayake KN,Kristaryanto Y,Jones RA,Vogt J,Rivagorda M,Handley MT,Mali GR,Quidwai T,Soares DC,Keighren MA,McKie L,Mort RL,Gammoh N,Garcia-Munoz A,Davey T,Vermeren M,Walsh D,Budd P,Aligianis IA,Faqeih E,Quigley AJ,Jackson IJ,Kulathu Y,Jackson M,Ribchester RR,von Kriegsheim A,Alkuraya FS,Woods CG,Maher ER,Mill P
Thu May 04 00:00:00 UTC 2017
PLAA Mutations Cause a Lethal Infantile Epileptic Encephalopathy by Disrupting Ubiquitin-Mediated Endolysosomal Degradation of Synaptic Proteins.
American journal of human genetics
Hall EA,Nahorski MS,Murray LM,Shaheen R,Perkins E,Dissanayake KN,Kristaryanto Y,Jones RA,Vogt J,Rivagorda M,Handley MT,Mali GR,Quidwai T,Soares DC,Keighren MA,McKie L,Mort RL,Gammoh N,Garcia-Munoz A,Davey T,Vermeren M,Walsh D,Budd P,Aligianis IA,Faqeih E,Quigley AJ,Jackson IJ,Kulathu Y,Jackson M,Ribchester RR,von Kriegsheim A,Alkuraya FS,Woods CG,Maher ER,Mill P
Thu May 04 00:00:00 UTC 2017
Dlitiazem inhibits the oxidative stress induced by angiotensin II through growth hormone secretagogue receptor type 1a in human umbilicus vein endothelial cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhou L,Yang M,Zuo S,Guan X,Wang J,Chen Q,Zuo X,Jia S,Guo R
Mon May 01 00:00:00 UTC 2017
Dlitiazem inhibits the oxidative stress induced by angiotensin II through growth hormone secretagogue receptor type 1a in human umbilicus vein endothelial cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhou L,Yang M,Zuo S,Guan X,Wang J,Chen Q,Zuo X,Jia S,Guo R
Mon May 01 00:00:00 UTC 2017
Protective effects of aloperine on monocrotaline-induced pulmonary hypertension in rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wu F,Hao Y,Yang J,Yao W,Xu Y,Yan L,Niu Y,Sun T,Yu J,Zhou R
Mon May 01 00:00:00 UTC 2017
Protective effects of aloperine on monocrotaline-induced pulmonary hypertension in rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wu F,Hao Y,Yang J,Yao W,Xu Y,Yan L,Niu Y,Sun T,Yu J,Zhou R
Mon May 01 00:00:00 UTC 2017
Long non-coding RNA TUG1 promotes cell proliferation and metastasis by negatively regulating miR-300 in gallbladder carcinoma.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ma F,Wang SH,Cai Q,Jin LY,Zhou D,Ding J,Quan ZW
Sat Apr 01 00:00:00 UTC 2017
Long non-coding RNA TUG1 promotes cell proliferation and metastasis by negatively regulating miR-300 in gallbladder carcinoma.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Ma F,Wang SH,Cai Q,Jin LY,Zhou D,Ding J,Quan ZW
Sat Apr 01 00:00:00 UTC 2017
Pretreatment Donors after Circulatory Death with Simvastatin Alleviates Liver Ischemia Reperfusion Injury through a KLF2-Dependent Mechanism in Rat.
Oxidative medicine and cellular longevity
Liu Z,Zhang X,Xiao Q,Ye S,Lai CH,Luo J,Huang X,Wang W,Zeng C,Zhong Z,Fan X,Xia Z,Xiong Y,Mao X,Ye Q,Wang Y
Fri Aug 17 00:00:00 UTC 2018
Pretreatment Donors after Circulatory Death with Simvastatin Alleviates Liver Ischemia Reperfusion Injury through a KLF2-Dependent Mechanism in Rat.
Oxidative medicine and cellular longevity
Liu Z,Zhang X,Xiao Q,Ye S,Lai CH,Luo J,Huang X,Wang W,Zeng C,Zhong Z,Fan X,Xia Z,Xiong Y,Mao X,Ye Q,Wang Y
Fri Aug 17 00:00:00 UTC 2018
HMGB1-induced autophagy: a new pathway to maintain Treg function during chronic hepatitis B virus infection.
Clinical science (London, England : 1979)
Cheng LS,Li J,Liu Y,Wang FP,Wang SQ,She WM,Wu SD,Qi XL,Zhou YP,Jiang W
Wed Mar 01 00:00:00 UTC 2017
HMGB1-induced autophagy: a new pathway to maintain Treg function during chronic hepatitis B virus infection.
Clinical science (London, England : 1979)
Cheng LS,Li J,Liu Y,Wang FP,Wang SQ,She WM,Wu SD,Qi XL,Zhou YP,Jiang W
Wed Mar 01 00:00:00 UTC 2017
The Long Noncoding RNA NEAT1 Exerts Antihantaviral Effects by Acting as Positive Feedback for RIG-I Signaling.
Journal of virology
Ma H,Han P,Ye W,Chen H,Zheng X,Cheng L,Zhang L,Yu L,Wu X,Xu Z,Lei Y,Zhang F
Mon May 01 00:00:00 UTC 2017
The Long Noncoding RNA NEAT1 Exerts Antihantaviral Effects by Acting as Positive Feedback for RIG-I Signaling.
Journal of virology
Ma H,Han P,Ye W,Chen H,Zheng X,Cheng L,Zhang L,Yu L,Wu X,Xu Z,Lei Y,Zhang F
Mon May 01 00:00:00 UTC 2017
The Circadian Gene Clock Regulates Bone Formation Via PDIA3.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Yuan G,Hua B,Yang Y,Xu L,Cai T,Sun N,Yan Z,Lu C,Qian R
Sat Apr 01 00:00:00 UTC 2017
The Circadian Gene Clock Regulates Bone Formation Via PDIA3.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Yuan G,Hua B,Yang Y,Xu L,Cai T,Sun N,Yan Z,Lu C,Qian R
Sat Apr 01 00:00:00 UTC 2017
Inhibition of HIF-1α by PX-478 suppresses tumor growth of esophageal squamous cell cancer in vitro and in vivo.
American journal of cancer research
Zhu Y,Zang Y,Zhao F,Li Z,Zhang J,Fang L,Li M,Xing L,Xu Z,Yu J
Mon Mar 21 00:00:00 UTC 2022
Inhibition of HIF-1α by PX-478 suppresses tumor growth of esophageal squamous cell cancer in vitro and in vivo.
American journal of cancer research
Zhu Y,Zang Y,Zhao F,Li Z,Zhang J,Fang L,Li M,Xing L,Xu Z,Yu J
Mon Mar 21 00:00:00 UTC 2022
Whole exome-wide association study identifies a missense variant in SLC2A4RG associated with glioblastoma risk.
American journal of cancer research
Zhao Y,Yun D,Zou X,Jiang T,Li G,Hu L,Chen J,Xu J,Mao Y,Chen H,Lu D
Wed Sep 30 00:00:00 UTC 2020
Whole exome-wide association study identifies a missense variant in SLC2A4RG associated with glioblastoma risk.
American journal of cancer research
Zhao Y,Yun D,Zou X,Jiang T,Li G,Hu L,Chen J,Xu J,Mao Y,Chen H,Lu D
Wed Sep 30 00:00:00 UTC 2020
Phosphocreatine protects endothelial cells from Methylglyoxal induced oxidative stress and apoptosis via the regulation of PI3K/Akt/eNOS and NF-κB pathway.
Vascular pharmacology
Chu P,Han G,Ahsan A,Sun Z,Liu S,Zhang Z,Sun B,Song Y,Lin Y,Peng J,Tang Z
Sat Apr 01 00:00:00 UTC 2017
Phosphocreatine protects endothelial cells from Methylglyoxal induced oxidative stress and apoptosis via the regulation of PI3K/Akt/eNOS and NF-κB pathway.
Vascular pharmacology
Chu P,Han G,Ahsan A,Sun Z,Liu S,Zhang Z,Sun B,Song Y,Lin Y,Peng J,Tang Z
Sat Apr 01 00:00:00 UTC 2017
AGR2 ameliorates tumor necrosis factor-α-induced epithelial barrier dysfunction via suppression of NF-κB p65-mediated MLCK/p-MLC pathway activation.
International journal of molecular medicine
Ye X,Sun M
Mon May 01 00:00:00 UTC 2017
AGR2 ameliorates tumor necrosis factor-α-induced epithelial barrier dysfunction via suppression of NF-κB p65-mediated MLCK/p-MLC pathway activation.
International journal of molecular medicine
Ye X,Sun M
Mon May 01 00:00:00 UTC 2017
Electroacupuncture Improved Hippocampal Neurogenesis following Traumatic Brain Injury in Mice through Inhibition of TLR4 Signaling Pathway.
Stem cells international
Ye Y,Yang Y,Chen C,Li Z,Jia Y,Su X,Wang C,He X
Tue Sep 29 00:00:00 UTC 2020
Electroacupuncture Improved Hippocampal Neurogenesis following Traumatic Brain Injury in Mice through Inhibition of TLR4 Signaling Pathway.
Stem cells international
Ye Y,Yang Y,Chen C,Li Z,Jia Y,Su X,Wang C,He X
Tue Sep 29 00:00:00 UTC 2020
ApoA-I/SR-BI modulates S1P/S1PR2-mediated inflammation through the PI3K/Akt signaling pathway in HUVECs.
Journal of physiology and biochemistry
Ren K,Lu YJ,Mo ZC,-Liu X,Tang ZL,Jiang Y,Peng XS,Li L,Zhang QH,Yi GH
Mon May 01 00:00:00 UTC 2017
ApoA-I/SR-BI modulates S1P/S1PR2-mediated inflammation through the PI3K/Akt signaling pathway in HUVECs.
Journal of physiology and biochemistry
Ren K,Lu YJ,Mo ZC,-Liu X,Tang ZL,Jiang Y,Peng XS,Li L,Zhang QH,Yi GH
Mon May 01 00:00:00 UTC 2017
Estrogen receptor-α36 is involved in icaritin induced growth inhibition of triple-negative breast cancer cells.
The Journal of steroid biochemistry and molecular biology
Wang X,Zheng N,Dong J,Wang X,Liu L,Huang J
Sat Jul 01 00:00:00 UTC 2017
Estrogen receptor-α36 is involved in icaritin induced growth inhibition of triple-negative breast cancer cells.
The Journal of steroid biochemistry and molecular biology
Wang X,Zheng N,Dong J,Wang X,Liu L,Huang J
Sat Jul 01 00:00:00 UTC 2017
1,2-Dichloroethane impairs glucose and lipid homeostasis in the livers of NIH Swiss mice.
Toxicology
Wang T,Xu D,Fan Q,Rong W,Zheng J,Gao C,Li G,Zeng N,Guo T,Zeng L,Wang F,Xiao C,Cai L,Tang S,Deng X,Yin X,Huang M,Lu F,Hu Q,Chen W,Huang Z,Wang Q
Sat Apr 01 00:00:00 UTC 2017
1,2-Dichloroethane impairs glucose and lipid homeostasis in the livers of NIH Swiss mice.
Toxicology
Wang T,Xu D,Fan Q,Rong W,Zheng J,Gao C,Li G,Zeng N,Guo T,Zeng L,Wang F,Xiao C,Cai L,Tang S,Deng X,Yin X,Huang M,Lu F,Hu Q,Chen W,Huang Z,Wang Q
Sat Apr 01 00:00:00 UTC 2017
The acute exposure of tetrachloro-p-benzoquinone (a.k.a. chloranil) triggers inflammation and neurological dysfunction via Toll-like receptor 4 signaling: The protective role of melatonin preconditioning.
Toxicology
Fu J,Xia X,Liu Z,Wang Y,Wang Y,Shi Q,Song X,Song E,Song Y
Sat Apr 15 00:00:00 UTC 2017
The acute exposure of tetrachloro-p-benzoquinone (a.k.a. chloranil) triggers inflammation and neurological dysfunction via Toll-like receptor 4 signaling: The protective role of melatonin preconditioning.
Toxicology
Fu J,Xia X,Liu Z,Wang Y,Wang Y,Shi Q,Song X,Song E,Song Y
Sat Apr 15 00:00:00 UTC 2017
β-Caryophyllene Pretreatment Alleviates Focal Cerebral Ischemia-Reperfusion Injury by Activating PI3K/Akt Signaling Pathway.
Neurochemical research
Zhang Q,An R,Tian X,Yang M,Li M,Lou J,Xu L,Dong Z
Mon May 01 00:00:00 UTC 2017
β-Caryophyllene Pretreatment Alleviates Focal Cerebral Ischemia-Reperfusion Injury by Activating PI3K/Akt Signaling Pathway.
Neurochemical research
Zhang Q,An R,Tian X,Yang M,Li M,Lou J,Xu L,Dong Z
Mon May 01 00:00:00 UTC 2017
Inhibition of Calcineurin A by FK506 Suppresses Seizures and Reduces the Expression of GluN2B in Membrane Fraction.
Neurochemical research
Wen Y,Fu P,Wu K,Si K,Xie Y,Dan W,Zhan Y,Shi Q
Tue Aug 01 00:00:00 UTC 2017
Inhibition of Calcineurin A by FK506 Suppresses Seizures and Reduces the Expression of GluN2B in Membrane Fraction.
Neurochemical research
Wen Y,Fu P,Wu K,Si K,Xie Y,Dan W,Zhan Y,Shi Q
Tue Aug 01 00:00:00 UTC 2017
Neuroprotective Effect of Chitosan Oligosaccharide on Hypoxic-Ischemic Brain Damage in Neonatal Rats.
Neurochemical research
Wu W,Wei W,Lu M,Zhu X,Liu N,Niu Y,Sun T,Li Y,Yu J
Wed Nov 01 00:00:00 UTC 2017
Neuroprotective Effect of Chitosan Oligosaccharide on Hypoxic-Ischemic Brain Damage in Neonatal Rats.
Neurochemical research
Wu W,Wei W,Lu M,Zhu X,Liu N,Niu Y,Sun T,Li Y,Yu J
Wed Nov 01 00:00:00 UTC 2017
The Anticonvulsant and Neuroprotective Effects of Oxysophocarpine on Pilocarpine-Induced Convulsions in Adult Male Mice.
Cellular and molecular neurobiology
Liu G,Wang J,Deng XH,Ma PS,Li FM,Peng XD,Niu Y,Sun T,Li YX,Yu JQ
Wed Mar 01 00:00:00 UTC 2017
The Anticonvulsant and Neuroprotective Effects of Oxysophocarpine on Pilocarpine-Induced Convulsions in Adult Male Mice.
Cellular and molecular neurobiology
Liu G,Wang J,Deng XH,Ma PS,Li FM,Peng XD,Niu Y,Sun T,Li YX,Yu JQ
Wed Mar 01 00:00:00 UTC 2017
miR-367 promotes tumor growth by inhibiting FBXW7 in NSCLC.
Oncology reports
Xiao G,Gao X,Sun X,Yang C,Zhang B,Sun R,Huang G,Li X,Liu J,Du N,Liu D,Liang R,Ren H,Qin S
Tue Aug 01 00:00:00 UTC 2017
miR-367 promotes tumor growth by inhibiting FBXW7 in NSCLC.
Oncology reports
Xiao G,Gao X,Sun X,Yang C,Zhang B,Sun R,Huang G,Li X,Liu J,Du N,Liu D,Liang R,Ren H,Qin S
Tue Aug 01 00:00:00 UTC 2017
BMP4 promotes mouse iPS cell differentiation to male germ cells via Smad1/5, Gata4, Id1 and Id2.
Reproduction (Cambridge, England)
Yang S,Yuan Q,Niu M,Hou J,Zhu Z,Sun M,Li Z,He Z
Wed Feb 01 00:00:00 UTC 2017
BMP4 promotes mouse iPS cell differentiation to male germ cells via Smad1/5, Gata4, Id1 and Id2.
Reproduction (Cambridge, England)
Yang S,Yuan Q,Niu M,Hou J,Zhu Z,Sun M,Li Z,He Z
Wed Feb 01 00:00:00 UTC 2017
Relationship between abnormal TOX3 gene methylation and polycystic ovarian syndrome.
European review for medical and pharmacological sciences
Ning Z,Jiayi L,Jian R,Wanli X
Mon May 01 00:00:00 UTC 2017
Relationship between abnormal TOX3 gene methylation and polycystic ovarian syndrome.
European review for medical and pharmacological sciences
Ning Z,Jiayi L,Jian R,Wanli X
Mon May 01 00:00:00 UTC 2017
A novel role of fragile X mental retardation protein in pre-mRNA alternative splicing through RNA-binding protein 14.
Neuroscience
Zhou LT,Ye SH,Yang HX,Zhou YT,Zhao QH,Sun WW,Gao MM,Yi YH,Long YS
Thu May 04 00:00:00 UTC 2017
A novel role of fragile X mental retardation protein in pre-mRNA alternative splicing through RNA-binding protein 14.
Neuroscience
Zhou LT,Ye SH,Yang HX,Zhou YT,Zhao QH,Sun WW,Gao MM,Yi YH,Long YS
Thu May 04 00:00:00 UTC 2017
Effects of hydrogen sulfide on inducible nitric oxide synthase activity and expression of cardiomyocytes in diabetic rats.
Molecular medicine reports
Yang R,Jia Q,Liu XF,Wang YY,Ma SF
Sun Oct 01 00:00:00 UTC 2017
Effects of hydrogen sulfide on inducible nitric oxide synthase activity and expression of cardiomyocytes in diabetic rats.
Molecular medicine reports
Yang R,Jia Q,Liu XF,Wang YY,Ma SF
Sun Oct 01 00:00:00 UTC 2017
Generation and characterization of induced pluripotent stem cells from guinea pig fetal fibroblasts.
Molecular medicine reports
Wu Y,Li O,He C,Li Y,Li M,Liu XL,Wang Y,He Y
Thu Jun 01 00:00:00 UTC 2017
Generation and characterization of induced pluripotent stem cells from guinea pig fetal fibroblasts.
Molecular medicine reports
Wu Y,Li O,He C,Li Y,Li M,Liu XL,Wang Y,He Y
Thu Jun 01 00:00:00 UTC 2017
CMIP is oncogenic in human gastric cancer cells.
Molecular medicine reports
Zhang J,Huang J,Wang X,Chen W,Tang Q,Fang M,Qian Y
Wed Nov 01 00:00:00 UTC 2017
CMIP is oncogenic in human gastric cancer cells.
Molecular medicine reports
Zhang J,Huang J,Wang X,Chen W,Tang Q,Fang M,Qian Y
Wed Nov 01 00:00:00 UTC 2017
Icariin inhibits MMP‑1, MMP‑3 and MMP‑13 expression through MAPK pathways in IL‑1β‑stimulated SW1353 chondrosarcoma cells.
Molecular medicine reports
Zeng L,Rong XF,Li RH,Wu XY
Mon May 01 00:00:00 UTC 2017
Icariin inhibits MMP‑1, MMP‑3 and MMP‑13 expression through MAPK pathways in IL‑1β‑stimulated SW1353 chondrosarcoma cells.
Molecular medicine reports
Zeng L,Rong XF,Li RH,Wu XY
Mon May 01 00:00:00 UTC 2017
Antitumor effects of baicalin on ovarian cancer cells through induction of cell apoptosis and inhibition of cell migration in vitro.
Molecular medicine reports
Gao C,Zhou Y,Li H,Cong X,Jiang Z,Wang X,Cao R,Tian W
Fri Dec 01 00:00:00 UTC 2017
Antitumor effects of baicalin on ovarian cancer cells through induction of cell apoptosis and inhibition of cell migration in vitro.
Molecular medicine reports
Gao C,Zhou Y,Li H,Cong X,Jiang Z,Wang X,Cao R,Tian W
Fri Dec 01 00:00:00 UTC 2017
SHIP, a novel factor to ameliorate extracellular matrix accumulation via suppressing PI3K/Akt/CTGF signaling in diabetic kidney disease.
Biochemical and biophysical research communications
Li F,Li L,Cheng M,Wang X,Hao J,Liu S,Duan H
Sun Jan 22 00:00:00 UTC 2017
SHIP, a novel factor to ameliorate extracellular matrix accumulation via suppressing PI3K/Akt/CTGF signaling in diabetic kidney disease.
Biochemical and biophysical research communications
Li F,Li L,Cheng M,Wang X,Hao J,Liu S,Duan H
Sun Jan 22 00:00:00 UTC 2017
A high-fat, high-protein diet attenuates the negative impact of casein-induced chronic inflammation on testicular steroidogenesis and sperm parameters in adult mice.
General and comparative endocrinology
Zhao JL,Zhao YY,Zhu WJ
Sun Oct 01 00:00:00 UTC 2017
A high-fat, high-protein diet attenuates the negative impact of casein-induced chronic inflammation on testicular steroidogenesis and sperm parameters in adult mice.
General and comparative endocrinology
Zhao JL,Zhao YY,Zhu WJ
Sun Oct 01 00:00:00 UTC 2017
Role of autophagy in cadmium-induced testicular injury.
Human & experimental toxicology
Wang YJ,Yan J,Yin F,Li L,Qin YG,Meng CY,Lu RF,Guo L
Sun Oct 01 00:00:00 UTC 2017
Role of autophagy in cadmium-induced testicular injury.
Human & experimental toxicology
Wang YJ,Yan J,Yin F,Li L,Qin YG,Meng CY,Lu RF,Guo L
Sun Oct 01 00:00:00 UTC 2017
Chloroform extract of Hedyotis diffusa Willd inhibits viability of human colorectal cancer cells via suppression of AKT and ERK signaling pathways.
Oncology letters
Yan Z,Feng J,Peng J,Lai Z,Zhang L,Jin Y,Yang H,Chen W,Lin J
Fri Dec 01 00:00:00 UTC 2017
Chloroform extract of Hedyotis diffusa Willd inhibits viability of human colorectal cancer cells via suppression of AKT and ERK signaling pathways.
Oncology letters
Yan Z,Feng J,Peng J,Lai Z,Zhang L,Jin Y,Yang H,Chen W,Lin J
Fri Dec 01 00:00:00 UTC 2017
Terrein performs antitumor functions on esophageal cancer cells by inhibiting cell proliferation and synergistic interaction with cisplatin.
Oncology letters
Wu Y,Zhu Y,Li S,Zeng M,Chu J,Hu P,Li J,Guo Q,Lv XB,Huang G
Sat Apr 01 00:00:00 UTC 2017
Terrein performs antitumor functions on esophageal cancer cells by inhibiting cell proliferation and synergistic interaction with cisplatin.
Oncology letters
Wu Y,Zhu Y,Li S,Zeng M,Chu J,Hu P,Li J,Guo Q,Lv XB,Huang G
Sat Apr 01 00:00:00 UTC 2017
miR-150 is downregulated in osteosarcoma and suppresses cell proliferation, migration and invasion by targeting ROCK1.
Oncology letters
Li CH,Yu TB,Qiu HW,Zhao X,Zhou CL,Qi C
Sat Apr 01 00:00:00 UTC 2017
miR-150 is downregulated in osteosarcoma and suppresses cell proliferation, migration and invasion by targeting ROCK1.
Oncology letters
Li CH,Yu TB,Qiu HW,Zhao X,Zhou CL,Qi C
Sat Apr 01 00:00:00 UTC 2017
Exosomes derived from bone marrow stromal cells decrease the sensitivity of leukemic cells to etoposide.
Oncology letters
Wang J,Li D,Zhuang Y,Fu J,Li X,Shi Q,Ju X
Fri Sep 01 00:00:00 UTC 2017
Exosomes derived from bone marrow stromal cells decrease the sensitivity of leukemic cells to etoposide.
Oncology letters
Wang J,Li D,Zhuang Y,Fu J,Li X,Shi Q,Ju X
Fri Sep 01 00:00:00 UTC 2017
Inhibition of chemotherapy-induced apoptosis of testicular cells by squid ink polysaccharide.
Experimental and therapeutic medicine
Gu YP,Yang XM,Duan ZH,Luo P,Shang JH,Xiao W,Tao YX,Zhang DY,Zhang YB,Liu HZ
Fri Dec 01 00:00:00 UTC 2017
Inhibition of chemotherapy-induced apoptosis of testicular cells by squid ink polysaccharide.
Experimental and therapeutic medicine
Gu YP,Yang XM,Duan ZH,Luo P,Shang JH,Xiao W,Tao YX,Zhang DY,Zhang YB,Liu HZ
Fri Dec 01 00:00:00 UTC 2017
Antiplatelet drug ticagrelor delays gastric ulcer healing in rats.
Experimental and therapeutic medicine
Li JJ,Wu XY,Chen JL,Chen GR,Xu J,Gu Y,Song HP
Sun Oct 01 00:00:00 UTC 2017
Antiplatelet drug ticagrelor delays gastric ulcer healing in rats.
Experimental and therapeutic medicine
Li JJ,Wu XY,Chen JL,Chen GR,Xu J,Gu Y,Song HP
Sun Oct 01 00:00:00 UTC 2017
Roux-en-Y gastric bypass surgery suppresses hypothalamic PTP1B protein level and alleviates leptin resistance in obese rats.
Experimental and therapeutic medicine
Liu JY,Mu S,Zhang SP,Guo W,Li QF,Xiao XQ,Zhang J,Wang ZH
Fri Sep 01 00:00:00 UTC 2017
Roux-en-Y gastric bypass surgery suppresses hypothalamic PTP1B protein level and alleviates leptin resistance in obese rats.
Experimental and therapeutic medicine
Liu JY,Mu S,Zhang SP,Guo W,Li QF,Xiao XQ,Zhang J,Wang ZH
Fri Sep 01 00:00:00 UTC 2017
DDX50 inhibits the replication of dengue virus 2 by upregulating IFN-β production.
Archives of virology
Han P,Ye W,Lv X,Ma H,Weng D,Dong Y,Cheng L,Chen H,Zhang L,Xu Z,Lei Y,Zhang F
Thu Jun 01 00:00:00 UTC 2017
DDX50 inhibits the replication of dengue virus 2 by upregulating IFN-β production.
Archives of virology
Han P,Ye W,Lv X,Ma H,Weng D,Dong Y,Cheng L,Chen H,Zhang L,Xu Z,Lei Y,Zhang F
Thu Jun 01 00:00:00 UTC 2017
Chondroprotective Effects and Multitarget Mechanisms of Fu Yuan Capsule in a Rat Osteoarthritis Model.
Evidence-based complementary and alternative medicine : eCAM
Zeng L,Xiao CZ,Deng ZT,Li RH
Thu Oct 01 00:00:00 UTC 2020
Chondroprotective Effects and Multitarget Mechanisms of Fu Yuan Capsule in a Rat Osteoarthritis Model.
Evidence-based complementary and alternative medicine : eCAM
Zeng L,Xiao CZ,Deng ZT,Li RH
Thu Oct 01 00:00:00 UTC 2020
Hyperlipidemia impairs erectile function in rats by causing cavernosal fibrosis.
Andrologia
Li R,Cui K,Wang T,Wang S,Li X,Qiu J,Yu G,Liu J,Wen B,Rao K
Fri Sep 01 00:00:00 UTC 2017
Hyperlipidemia impairs erectile function in rats by causing cavernosal fibrosis.
Andrologia
Li R,Cui K,Wang T,Wang S,Li X,Qiu J,Yu G,Liu J,Wen B,Rao K
Fri Sep 01 00:00:00 UTC 2017
Esculetin induces apoptosis of SMMC-7721 cells through IGF-1/PI3K/Akt-mediated mitochondrial pathways.
Canadian journal of physiology and pharmacology
Li J,Li S,Wang X,Wang H
Sat Jul 01 00:00:00 UTC 2017
Esculetin induces apoptosis of SMMC-7721 cells through IGF-1/PI3K/Akt-mediated mitochondrial pathways.
Canadian journal of physiology and pharmacology
Li J,Li S,Wang X,Wang H
Sat Jul 01 00:00:00 UTC 2017
Mitochondrial dynamics changes with age in an APPsw/PS1dE9 mouse model of Alzheimer's disease.
Neuroreport
Xu LL,Shen Y,Wang X,Wei LF,Wang P,Yang H,Wang CF,Xie ZH,Bi JZ
Wed Mar 01 00:00:00 UTC 2017
Mitochondrial dynamics changes with age in an APPsw/PS1dE9 mouse model of Alzheimer's disease.
Neuroreport
Xu LL,Shen Y,Wang X,Wei LF,Wang P,Yang H,Wang CF,Xie ZH,Bi JZ
Wed Mar 01 00:00:00 UTC 2017
Functional characterization of a novel transcript of ERCC1 in chemotherapy resistance of ovarian cancer.
Oncotarget
Liu J,Zhang L,Mao P,Jiang G,Liu L,Wang J,Yang W,Owusu L,Li W
Tue Oct 17 00:00:00 UTC 2017
Functional characterization of a novel transcript of ERCC1 in chemotherapy resistance of ovarian cancer.
Oncotarget
Liu J,Zhang L,Mao P,Jiang G,Liu L,Wang J,Yang W,Owusu L,Li W
Tue Oct 17 00:00:00 UTC 2017
Coptisine from Rhizoma Coptidis Suppresses HCT-116 Cells-related Tumor Growth in vitro and in vivo.
Scientific reports
Huang T,Xiao Y,Yi L,Li L,Wang M,Tian C,Ma H,He K,Wang Y,Han B,Ye X,Li X
Mon Feb 06 00:00:00 UTC 2017
Coptisine from Rhizoma Coptidis Suppresses HCT-116 Cells-related Tumor Growth in vitro and in vivo.
Scientific reports
Huang T,Xiao Y,Yi L,Li L,Wang M,Tian C,Ma H,He K,Wang Y,Han B,Ye X,Li X
Mon Feb 06 00:00:00 UTC 2017
Hepatoprotective effect of quercetin against LPS/d-GalN induced acute liver injury in mice by inhibiting the IKK/NF-κB and MAPK signal pathways.
International immunopharmacology
Peng Z,Gong X,Yang Y,Huang L,Zhang Q,Zhang P,Wan R,Zhang B
Wed Nov 01 00:00:00 UTC 2017
Up-regulated BAFF and BAFF receptor expression in patients with intractable temporal lobe epilepsy and a pilocarpine-induced epilepsy rat model.
Seizure
Ma L,Li R,Huang H,Yuan J,Ou S,Xu T,Yu X,Liu X,Chen Y
Mon May 01 00:00:00 UTC 2017
Rosiglitazone Inhibits Angiotensin II-Induced Proliferation of Glomerular Mesangial Cells via the Gαq/Plcβ4/TRPC Signaling Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wei L,Mao J,Lu J,Gao J,Zhu D,Tian L,Chen Z,Jia L,Wang L,Fu R
Mon Mar 12 00:00:00 UTC 2018
Rosiglitazone Inhibits Angiotensin II-Induced Proliferation of Glomerular Mesangial Cells via the Gαq/Plcβ4/TRPC Signaling Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wei L,Mao J,Lu J,Gao J,Zhu D,Tian L,Chen Z,Jia L,Wang L,Fu R
Mon Mar 12 00:00:00 UTC 2018
A novel protein derived from lamprey supraneural body tissue with efficient cytocidal actions against tumor cells.
Cell communication and signaling : CCS
Pang Y,Li C,Wang S,Ba W,Yu T,Pei G,Bi D,Liang H,Pan X,Zhu T,Gou M,Han Y,Li Q
Mon Oct 16 00:00:00 UTC 2017
A novel protein derived from lamprey supraneural body tissue with efficient cytocidal actions against tumor cells.
Cell communication and signaling : CCS
Pang Y,Li C,Wang S,Ba W,Yu T,Pei G,Bi D,Liang H,Pan X,Zhu T,Gou M,Han Y,Li Q
Mon Oct 16 00:00:00 UTC 2017
KIAA1199 is induced by inflammation and enhances malignant phenotype in pancreatic cancer.
Oncotarget
Kohi S,Sato N,Koga A,Matayoshi N,Hirata K
Tue Mar 07 00:00:00 UTC 2017
KIAA1199 is induced by inflammation and enhances malignant phenotype in pancreatic cancer.
Oncotarget
Kohi S,Sato N,Koga A,Matayoshi N,Hirata K
Tue Mar 07 00:00:00 UTC 2017
Hypoxia-inducible factor-1 alpha is involved in RIP-induced necroptosis caused by in vitro and in vivo ischemic brain injury.
Scientific reports
Yang XS,Yi TL,Zhang S,Xu ZW,Yu ZQ,Sun HT,Yang C,Tu Y,Cheng SX
Wed Jul 19 00:00:00 UTC 2017
Hypoxia-inducible factor-1 alpha is involved in RIP-induced necroptosis caused by in vitro and in vivo ischemic brain injury.
Scientific reports
Yang XS,Yi TL,Zhang S,Xu ZW,Yu ZQ,Sun HT,Yang C,Tu Y,Cheng SX
Wed Jul 19 00:00:00 UTC 2017
Inhibition of the TRPM2 and TRPV1 Channels through Hypericum perforatum in Sciatic Nerve Injury-induced Rats Demonstrates their Key Role in Apoptosis and Mitochondrial Oxidative Stress of Sciatic Nerve and Dorsal Root Ganglion.
Frontiers in physiology
Uslusoy F,Nazıroğlu M,Çiğ B
Thu Oct 01 00:00:00 UTC 2020
Inhibition of the TRPM2 and TRPV1 Channels through Hypericum perforatum in Sciatic Nerve Injury-induced Rats Demonstrates their Key Role in Apoptosis and Mitochondrial Oxidative Stress of Sciatic Nerve and Dorsal Root Ganglion.
Frontiers in physiology
Uslusoy F,Nazıroğlu M,Çiğ B
Thu Oct 01 00:00:00 UTC 2020
Induction of apoptosis by FFJ-5, a novel naphthoquinone compound, occurs via downregulation of PKM2 in A549 and HepG2 cells.
Oncology letters
Wei X,Li M,Ma M,Jia H,Zhang Y,Kang W,Wang T,Shi X
Wed Feb 01 00:00:00 UTC 2017
Low Frequency Magnetic Fields Induce Autophagy-associated Cell Death in Lung Cancer through miR-486-mediated Inhibition of Akt/mTOR Signaling Pathway.
Scientific reports
Xu Y,Wang Y,Yao A,Xu Z,Dou H,Shen S,Hou Y,Wang T
Mon Sep 18 00:00:00 UTC 2017
Low Frequency Magnetic Fields Induce Autophagy-associated Cell Death in Lung Cancer through miR-486-mediated Inhibition of Akt/mTOR Signaling Pathway.
Scientific reports
Xu Y,Wang Y,Yao A,Xu Z,Dou H,Shen S,Hou Y,Wang T
Mon Sep 18 00:00:00 UTC 2017
Generation and characteristics of human Sertoli cell line immortalized by overexpression of human telomerase.
Oncotarget
Wen L,Yuan Q,Sun M,Niu M,Wang H,Fu H,Zhou F,Yao C,Wang X,Li Z,He Z
Tue Mar 07 00:00:00 UTC 2017
Generation and characteristics of human Sertoli cell line immortalized by overexpression of human telomerase.
Oncotarget
Wen L,Yuan Q,Sun M,Niu M,Wang H,Fu H,Zhou F,Yao C,Wang X,Li Z,He Z
Tue Mar 07 00:00:00 UTC 2017
TRIM14 regulates cell proliferation and invasion in osteosarcoma via promotion of the AKT signaling pathway.
Scientific reports
Xu G,Guo Y,Xu D,Wang Y,Shen Y,Wang F,Lv Y,Song F,Jiang D,Zhang Y,Lou Y,Meng Y,Yang Y,Kang Y
Thu Feb 16 00:00:00 UTC 2017
TRIM14 regulates cell proliferation and invasion in osteosarcoma via promotion of the AKT signaling pathway.
Scientific reports
Xu G,Guo Y,Xu D,Wang Y,Shen Y,Wang F,Lv Y,Song F,Jiang D,Zhang Y,Lou Y,Meng Y,Yang Y,Kang Y
Thu Feb 16 00:00:00 UTC 2017
Differential inhibition of activity, activation and gene expression of MMP-9 in THP-1 cells by azithromycin and minocycline versus bortezomib: A comparative study.
PloS one
Vandooren J,Knoops S,Aldinucci Buzzo JL,Boon L,Martens E,Opdenakker G,Kolaczkowska E
Wed Aug 30 00:00:00 UTC 2017
Differential inhibition of activity, activation and gene expression of MMP-9 in THP-1 cells by azithromycin and minocycline versus bortezomib: A comparative study.
PloS one
Vandooren J,Knoops S,Aldinucci Buzzo JL,Boon L,Martens E,Opdenakker G,Kolaczkowska E
Wed Aug 30 00:00:00 UTC 2017
Mechanism of Lycium barbarum polysaccharides on primary cultured rat hippocampal neurons.
Cell and tissue research
Zhao P,Ma NT,Chang RY,Li YX,Hao YJ,Yang WL,Zheng J,Niu Y,Sun T,Yu JQ
Fri Sep 01 00:00:00 UTC 2017
Beneficial effects of Lycium barbarum polysaccharide on spermatogenesis by improving antioxidant activity and inhibiting apoptosis in streptozotocin-induced diabetic male mice.
Food & function
Shi GJ,Zheng J,Wu J,Qiao HQ,Chang Q,Niu Y,Sun T,Li YX,Yu JQ
Wed Mar 22 00:00:00 UTC 2017
HOTAIR regulates HK2 expression by binding endogenous miR-125 and miR-143 in oesophageal squamous cell carcinoma progression.
Oncotarget
Ma J,Fan Y,Feng T,Chen F,Xu Z,Li S,Lin Q,He X,Shi W,Liu Y,Liu Z,Zhu B,Cao X
Tue Oct 17 00:00:00 UTC 2017
HOTAIR regulates HK2 expression by binding endogenous miR-125 and miR-143 in oesophageal squamous cell carcinoma progression.
Oncotarget
Ma J,Fan Y,Feng T,Chen F,Xu Z,Li S,Lin Q,He X,Shi W,Liu Y,Liu Z,Zhu B,Cao X
Tue Oct 17 00:00:00 UTC 2017
Epigallocatechin-3-gallate inhibits H2O2-induced apoptosis in Mouse Vascular Smooth Muscle Cells via 67kD Laminin Receptor.
Scientific reports
Yan X,Li Y,Yu H,Wang W,Wu C,Yang Y,Hu Y,Shi X,Li J
Thu Aug 10 00:00:00 UTC 2017
Epigallocatechin-3-gallate inhibits H2O2-induced apoptosis in Mouse Vascular Smooth Muscle Cells via 67kD Laminin Receptor.
Scientific reports
Yan X,Li Y,Yu H,Wang W,Wu C,Yang Y,Hu Y,Shi X,Li J
Thu Aug 10 00:00:00 UTC 2017
LINC00461, a long non-coding RNA, is important for the proliferation and migration of glioma cells.
Oncotarget
Yang Y,Ren M,Song C,Li D,Soomro SH,Xiong Y,Zhang H,Fu H
Fri Oct 13 00:00:00 UTC 2017
Involvement of TRPM2 and TRPV1 channels on hyperalgesia, apoptosis and oxidative stress in rat fibromyalgia model: Protective role of selenium.
Scientific reports
Yüksel E,Nazıroğlu M,Şahin M,Çiğ B
Wed Dec 13 00:00:00 UTC 2017
Involvement of TRPM2 and TRPV1 channels on hyperalgesia, apoptosis and oxidative stress in rat fibromyalgia model: Protective role of selenium.
Scientific reports
Yüksel E,Nazıroğlu M,Şahin M,Çiğ B
Wed Dec 13 00:00:00 UTC 2017
Nrf2-Knockout Protects from Intestinal Injuries in C57BL/6J Mice Following Abdominal Irradiation with γ Rays.
International journal of molecular sciences
Yang W,Sun Z,Yang B,Wang Q
Mon Jul 31 00:00:00 UTC 2017
Nrf2-Knockout Protects from Intestinal Injuries in C57BL/6J Mice Following Abdominal Irradiation with γ Rays.
International journal of molecular sciences
Yang W,Sun Z,Yang B,Wang Q
Mon Jul 31 00:00:00 UTC 2017
Effect of Oleracein E, a Neuroprotective Tetrahydroisoquinoline, on Rotenone-Induced Parkinson's Disease Cell and Animal Models.
ACS chemical neuroscience
Sun H,He X,Liu C,Li L,Zhou R,Jin T,Yue S,Feng D,Gong J,Sun J,Ji J,Xiang L
Wed Jan 18 00:00:00 UTC 2017
Effect of Oleracein E, a Neuroprotective Tetrahydroisoquinoline, on Rotenone-Induced Parkinson's Disease Cell and Animal Models.
ACS chemical neuroscience
Sun H,He X,Liu C,Li L,Zhou R,Jin T,Yue S,Feng D,Gong J,Sun J,Ji J,Xiang L
Wed Jan 18 00:00:00 UTC 2017
IGFBP-rP1 acts as a potential tumor suppressor via the suppression of ERK signaling pathway in endometrial cancer cells.
Molecular medicine reports
Ma Y,Jiang J,Zhang Y,Ding Y,Xu T,Lu B
Tue Aug 01 00:00:00 UTC 2017
Boschniakia Rossica Polysaccharide Triggers Laryngeal Carcinoma Cell Apoptosis by Regulating Expression of Bcl-2, Caspase-3, and P53.
Medical science monitor : international medical journal of experimental and clinical research
Yao C,Cao X,Fu Z,Tian J,Dong W,Xu J,An K,Zhai L,Yu J
Sat Apr 29 00:00:00 UTC 2017
Boschniakia Rossica Polysaccharide Triggers Laryngeal Carcinoma Cell Apoptosis by Regulating Expression of Bcl-2, Caspase-3, and P53.
Medical science monitor : international medical journal of experimental and clinical research
Yao C,Cao X,Fu Z,Tian J,Dong W,Xu J,An K,Zhai L,Yu J
Sat Apr 29 00:00:00 UTC 2017
Melatonin ameliorates restraint stress-induced oxidative stress and apoptosis in testicular cells via NF-κB/iNOS and Nrf2/ HO-1 signaling pathway.
Scientific reports
Guo Y,Sun J,Li T,Zhang Q,Bu S,Wang Q,Lai D
Tue Aug 29 00:00:00 UTC 2017
Melatonin ameliorates restraint stress-induced oxidative stress and apoptosis in testicular cells via NF-κB/iNOS and Nrf2/ HO-1 signaling pathway.
Scientific reports
Guo Y,Sun J,Li T,Zhang Q,Bu S,Wang Q,Lai D
Tue Aug 29 00:00:00 UTC 2017
Tumor-associated macrophages promote Ezrin phosphorylation-mediated epithelial-mesenchymal transition in lung adenocarcinoma through FUT4/LeY up-regulation.
Oncotarget
Wang A,Lu C,Ning Z,Gao W,Xie Y,Zhang N,Liang J,Abbasi FS,Yan Q,Liu J
Tue Apr 25 00:00:00 UTC 2017
Tumor-associated macrophages promote Ezrin phosphorylation-mediated epithelial-mesenchymal transition in lung adenocarcinoma through FUT4/LeY up-regulation.
Oncotarget
Wang A,Lu C,Ning Z,Gao W,Xie Y,Zhang N,Liang J,Abbasi FS,Yan Q,Liu J
Tue Apr 25 00:00:00 UTC 2017
TGF-β1 promotes human hepatic carcinoma HepG2 cells invasion by upregulating autophagy.
European review for medical and pharmacological sciences
Ma CL,Qiao S,Li YC,Wang XF,Sun RJ,Zhang X,Qian RK,Song SD
Thu Jun 01 00:00:00 UTC 2017
Volatile Oil from Amomi Fructus Attenuates 5-Fluorouracil-Induced Intestinal Mucositis.
Frontiers in pharmacology
Zhang T,Lu SH,Bi Q,Liang L,Wang YF,Yang XX,Gu W,Yu J
Tue Sep 29 00:00:00 UTC 2020
Volatile Oil from Amomi Fructus Attenuates 5-Fluorouracil-Induced Intestinal Mucositis.
Frontiers in pharmacology
Zhang T,Lu SH,Bi Q,Liang L,Wang YF,Yang XX,Gu W,Yu J
Tue Sep 29 00:00:00 UTC 2020
AMPK activation-dependent autophagy compromises oleanolic acid-induced cytotoxicity in human bladder cancer cells.
Oncotarget
Song Y,Zhang P,Sun Y,Li X,Chen L,Xiao Y,Xing Y
Fri Sep 15 00:00:00 UTC 2017
AMPK activation-dependent autophagy compromises oleanolic acid-induced cytotoxicity in human bladder cancer cells.
Oncotarget
Song Y,Zhang P,Sun Y,Li X,Chen L,Xiao Y,Xing Y
Fri Sep 15 00:00:00 UTC 2017
Neuroprotective effect of liquiritin against neuropathic pain induced by chronic constriction injury of the sciatic nerve in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang MT,Wang B,Jia YN,Liu N,Ma PS,Gong SS,Niu Y,Sun T,Li YX,Yu JQ
Wed Nov 01 00:00:00 UTC 2017
Enhanced tenogenic differentiation and tendon-like tissue formation by CHIP overexpression in tendon-derived stem cells.
Acta biochimica et biophysica Sinica
Han W,Chen L,Liu J,Guo A
Sat Apr 01 00:00:00 UTC 2017
The tumor suppressor PTEN has a critical role in antiviral innate immunity.
Nature immunology
Li S,Zhu M,Pan R,Fang T,Cao YY,Chen S,Zhao X,Lei CQ,Guo L,Chen Y,Li CM,Jokitalo E,Yin Y,Shu HB,Guo D
Tue Mar 01 00:00:00 UTC 2016
The tumor suppressor PTEN has a critical role in antiviral innate immunity.
Nature immunology
Li S,Zhu M,Pan R,Fang T,Cao YY,Chen S,Zhao X,Lei CQ,Guo L,Chen Y,Li CM,Jokitalo E,Yin Y,Shu HB,Guo D
Tue Mar 01 00:00:00 UTC 2016
State estimation and prediction using clustered particle filters.
Proceedings of the National Academy of Sciences of the United States of America
Lee Y,Majda AJ
Tue Dec 20 00:00:00 UTC 2016
State estimation and prediction using clustered particle filters.
Proceedings of the National Academy of Sciences of the United States of America
Lee Y,Majda AJ
Tue Dec 20 00:00:00 UTC 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
Tue Dec 20 00:00:00 UTC 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
Tue Dec 20 00:00:00 UTC 2016
Interaction between endogenous carbon monoxide and hydrogen sulfide in the mechanism of gastroprotection against acute aspirin-induced gastric damage.
Pharmacological research
Magierowski M,Magierowska K,Hubalewska-Mazgaj M,Adamski J,Bakalarz D,Sliwowski Z,Pajdo R,Kwiecien S,Brzozowski T
Thu Dec 01 00:00:00 UTC 2016
Interaction between endogenous carbon monoxide and hydrogen sulfide in the mechanism of gastroprotection against acute aspirin-induced gastric damage.
Pharmacological research
Magierowski M,Magierowska K,Hubalewska-Mazgaj M,Adamski J,Bakalarz D,Sliwowski Z,Pajdo R,Kwiecien S,Brzozowski T
Thu Dec 01 00:00:00 UTC 2016
Disrupting MALAT1/miR-200c sponge decreases invasion and migration in endometrioid endometrial carcinoma.
Cancer letters
Li Q,Zhang C,Chen R,Xiong H,Qiu F,Liu S,Zhang M,Wang F,Wang Y,Zhou X,Xiao G,Wang X,Jiang Q
Thu Dec 01 00:00:00 UTC 2016
Disrupting MALAT1/miR-200c sponge decreases invasion and migration in endometrioid endometrial carcinoma.
Cancer letters
Li Q,Zhang C,Chen R,Xiong H,Qiu F,Liu S,Zhang M,Wang F,Wang Y,Zhou X,Xiao G,Wang X,Jiang Q
Thu Dec 01 00:00:00 UTC 2016
Lipodystrophy and severe metabolic dysfunction in mice with adipose tissue-specific insulin receptor ablation.
Molecular metabolism
Qiang G,Whang Kong H,Xu S,Pham HA,Parlee SD,Burr AA,Gil V,Pang J,Hughes A,Gu X,Fantuzzi G,MacDougald OA,Liew CW
Fri Jul 01 00:00:00 UTC 2016
Lipodystrophy and severe metabolic dysfunction in mice with adipose tissue-specific insulin receptor ablation.
Molecular metabolism
Qiang G,Whang Kong H,Xu S,Pham HA,Parlee SD,Burr AA,Gil V,Pang J,Hughes A,Gu X,Fantuzzi G,MacDougald OA,Liew CW
Fri Jul 01 00:00:00 UTC 2016
Lactobacillus rhamnosus induced epithelial cell apoptosis, ameliorates inflammation and prevents colon cancer development in an animal model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gamallat Y,Meyiah A,Kuugbee ED,Hago AM,Chiwala G,Awadasseid A,Bamba D,Zhang X,Shang X,Luo F,Xin Y
Sat Oct 01 00:00:00 UTC 2016
Lactobacillus rhamnosus induced epithelial cell apoptosis, ameliorates inflammation and prevents colon cancer development in an animal model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Gamallat Y,Meyiah A,Kuugbee ED,Hago AM,Chiwala G,Awadasseid A,Bamba D,Zhang X,Shang X,Luo F,Xin Y
Sat Oct 01 00:00:00 UTC 2016
Myricetin ameliorates brain injury and neurological deficits via Nrf2 activation after experimental stroke in middle-aged rats.
Food & function
Wu S,Yue Y,Peng A,Zhang L,Xiang J,Cao X,Ding H,Yin S
Wed Jun 15 00:00:00 UTC 2016
Myricetin ameliorates brain injury and neurological deficits via Nrf2 activation after experimental stroke in middle-aged rats.
Food & function
Wu S,Yue Y,Peng A,Zhang L,Xiang J,Cao X,Ding H,Yin S
Wed Jun 15 00:00:00 UTC 2016
Radiation promotes epithelial-to-mesenchymal transition and invasion of pancreatic cancer cell by activating carcinoma-associated fibroblasts.
American journal of cancer research
Li D,Qu C,Ning Z,Wang H,Zang K,Zhuang L,Chen L,Wang P,Meng Z
Thu Oct 01 00:00:00 UTC 2020
Radiation promotes epithelial-to-mesenchymal transition and invasion of pancreatic cancer cell by activating carcinoma-associated fibroblasts.
American journal of cancer research
Li D,Qu C,Ning Z,Wang H,Zang K,Zhuang L,Chen L,Wang P,Meng Z
Thu Oct 01 00:00:00 UTC 2020
IL-17 intensifies IFN-γ-induced NOS2 upregulation in RAW 264.7 cells by further activating STAT1 and NF-κB.
International journal of molecular medicine
Gao Q,Liu Y,Wu Y,Zhao Q,Wang L,Gao S,Wen W,Zhang W,Guo N,Zhou J,Yuan Z
Mon Feb 01 00:00:00 UTC 2016
miR-223 enhances the sensitivity of non-small cell lung cancer cells to erlotinib by targeting the insulin-like growth factor-1 receptor.
International journal of molecular medicine
Zhao FY,Han J,Chen XW,Wang J,Wang XD,Sun JG,Chen ZT
Fri Jul 01 00:00:00 UTC 2016
miR-223 enhances the sensitivity of non-small cell lung cancer cells to erlotinib by targeting the insulin-like growth factor-1 receptor.
International journal of molecular medicine
Zhao FY,Han J,Chen XW,Wang J,Wang XD,Sun JG,Chen ZT
Fri Jul 01 00:00:00 UTC 2016
Progranulin deficiency leads to severe inflammation, lung injury and cell death in a mouse model of endotoxic shock.
Journal of cellular and molecular medicine
Yu Y,Xu X,Liu L,Mao S,Feng T,Lu Y,Cheng Y,Wang H,Zhao W,Tang W
Tue Mar 01 00:00:00 UTC 2016
Effect of acute lipopolysaccharide-induced inflammation in intracerebroventricular-streptozotocin injected rats.
Neuropharmacology
Murtishaw AS,Heaney CF,Bolton MM,Sabbagh JJ,Langhardt MA,Kinney JW
Mon Feb 01 00:00:00 UTC 2016
Effect of acute lipopolysaccharide-induced inflammation in intracerebroventricular-streptozotocin injected rats.
Neuropharmacology
Murtishaw AS,Heaney CF,Bolton MM,Sabbagh JJ,Langhardt MA,Kinney JW
Mon Feb 01 00:00:00 UTC 2016
Anti-tumor effects of osthole on ovarian cancer cells in vitro.
Journal of ethnopharmacology
Jiang G,Liu J,Ren B,Tang Y,Owusu L,Li M,Zhang J,Liu L,Li W
Sun Dec 04 00:00:00 UTC 2016
Anti-tumor effects of osthole on ovarian cancer cells in vitro.
Journal of ethnopharmacology
Jiang G,Liu J,Ren B,Tang Y,Owusu L,Li M,Zhang J,Liu L,Li W
Sun Dec 04 00:00:00 UTC 2016
Bezafibrate-mizoribine interaction: Involvement of organic anion transporters OAT1 and OAT3 in rats.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Feng Y,Wang C,Liu Q,Meng Q,Huo X,Liu Z,Sun P,Yang X,Sun H,Qin J,Liu K
Fri Jan 01 00:00:00 UTC 2016
Bezafibrate-mizoribine interaction: Involvement of organic anion transporters OAT1 and OAT3 in rats.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Feng Y,Wang C,Liu Q,Meng Q,Huo X,Liu Z,Sun P,Yang X,Sun H,Qin J,Liu K
Fri Jan 01 00:00:00 UTC 2016
ApoA-I induces S1P release from endothelial cells through ABCA1 and SR-BI in a positive feedback manner.
Journal of physiology and biochemistry
Liu X,Ren K,Suo R,Xiong SL,Zhang QH,Mo ZC,Tang ZL,Jiang Y,Peng XS,Yi GH
Thu Dec 01 00:00:00 UTC 2016
ApoA-I induces S1P release from endothelial cells through ABCA1 and SR-BI in a positive feedback manner.
Journal of physiology and biochemistry
Liu X,Ren K,Suo R,Xiong SL,Zhang QH,Mo ZC,Tang ZL,Jiang Y,Peng XS,Yi GH
Thu Dec 01 00:00:00 UTC 2016
The Generation and Characterization of Recombinant Protein and Antibodies of Clostridium perfringens Beta2 Toxin.
Journal of immunology research
Zeng J,Song F,Yang Y,Ma C,Deng G,Li Y,Wang Y,Liu X
Wed Nov 20 00:00:00 UTC 2019
The Generation and Characterization of Recombinant Protein and Antibodies of Clostridium perfringens Beta2 Toxin.
Journal of immunology research
Zeng J,Song F,Yang Y,Ma C,Deng G,Li Y,Wang Y,Liu X
Wed Nov 20 00:00:00 UTC 2019
Potassium supplementation inhibits IL-17A production induced by salt loading in human T lymphocytes via p38/MAPK-SGK1 pathway.
Experimental and molecular pathology
Wen W,Wan Z,Ren K,Zhou D,Gao Q,Wu Y,Wang L,Yuan Z,Zhou J
Wed Jun 01 00:00:00 UTC 2016
Potassium supplementation inhibits IL-17A production induced by salt loading in human T lymphocytes via p38/MAPK-SGK1 pathway.
Experimental and molecular pathology
Wen W,Wan Z,Ren K,Zhou D,Gao Q,Wu Y,Wang L,Yuan Z,Zhou J
Wed Jun 01 00:00:00 UTC 2016
High-dose irradiation in combination with toll-like receptor 9 agonist CpG oligodeoxynucleotide 7909 downregulates PD-L1 expression via the NF-κB signaling pathway in non-small cell lung cancer cells.
OncoTargets and therapy
Chen X,Zhang Q,Luo Y,Gao C,Zhuang X,Xu G,Qiao T
Thu Oct 01 00:00:00 UTC 2020
High-dose irradiation in combination with toll-like receptor 9 agonist CpG oligodeoxynucleotide 7909 downregulates PD-L1 expression via the NF-κB signaling pathway in non-small cell lung cancer cells.
OncoTargets and therapy
Chen X,Zhang Q,Luo Y,Gao C,Zhuang X,Xu G,Qiao T
Thu Oct 01 00:00:00 UTC 2020
Nicotine Induced Murine Spermatozoa Apoptosis via Up-Regulation of Deubiquitinated RIP1 by Trim27 Promoter Hypomethylation.
Biology of reproduction
Nie D,Zhang D,Dai J,Zhang M,Zhao X,Xu W,Chen Z,Wang L,Wang Z,Qiao Z
Mon Feb 01 00:00:00 UTC 2016
Nicotine Induced Murine Spermatozoa Apoptosis via Up-Regulation of Deubiquitinated RIP1 by Trim27 Promoter Hypomethylation.
Biology of reproduction
Nie D,Zhang D,Dai J,Zhang M,Zhao X,Xu W,Chen Z,Wang L,Wang Z,Qiao Z
Mon Feb 01 00:00:00 UTC 2016
The long noncoding RNA MALAT1 regulates the lipopolysaccharide-induced inflammatory response through its interaction with NF-κB.
FEBS letters
Zhao G,Su Z,Song D,Mao Y,Mao X
Thu Sep 01 00:00:00 UTC 2016
The long noncoding RNA MALAT1 regulates the lipopolysaccharide-induced inflammatory response through its interaction with NF-κB.
FEBS letters
Zhao G,Su Z,Song D,Mao Y,Mao X
Thu Sep 01 00:00:00 UTC 2016
miR-1273g-3p modulates activation and apoptosis of hepatic stellate cells by directly targeting PTEN in HCV-related liver fibrosis.
FEBS letters
Niu X,Fu N,Du J,Wang R,Wang Y,Zhao S,Du H,Wang B,Zhang Y,Sun D,Nan Y
Mon Aug 01 00:00:00 UTC 2016
miR-1273g-3p modulates activation and apoptosis of hepatic stellate cells by directly targeting PTEN in HCV-related liver fibrosis.
FEBS letters
Niu X,Fu N,Du J,Wang R,Wang Y,Zhao S,Du H,Wang B,Zhang Y,Sun D,Nan Y
Mon Aug 01 00:00:00 UTC 2016
Upregulated Expression of microRNA-16 Correlates with Th17/Treg Cell Imbalance in Patients with Rheumatoid Arthritis.
DNA and cell biology
Wu YH,Liu W,Xue B,Zhang L,Liu XY,Liu B,Wang Y,Cai Y,Duan R
Thu Dec 01 00:00:00 UTC 2016
Hydrogen Sulfide and Carbon Monoxide Protect Gastric Mucosa Compromised by Mild Stress Against Alendronate Injury.
Digestive diseases and sciences
Magierowski M,Magierowska K,Szmyd J,Surmiak M,Sliwowski Z,Kwiecien S,Brzozowski T
Tue Nov 01 00:00:00 UTC 2016
Hydrogen Sulfide and Carbon Monoxide Protect Gastric Mucosa Compromised by Mild Stress Against Alendronate Injury.
Digestive diseases and sciences
Magierowski M,Magierowska K,Szmyd J,Surmiak M,Sliwowski Z,Kwiecien S,Brzozowski T
Tue Nov 01 00:00:00 UTC 2016
[Corrigendum] Cannabinoid WIN55, 212-2 induces cell cycle arrest and inhibits the proliferation and migration of human BEL7402 hepatocellular carcinoma cells.
Molecular medicine reports
Xu D,Wang J,Zhou Z,He Z,Zhao Q
Fri Jan 01 00:00:00 UTC 2016
[Corrigendum] Cannabinoid WIN55, 212-2 induces cell cycle arrest and inhibits the proliferation and migration of human BEL7402 hepatocellular carcinoma cells.
Molecular medicine reports
Xu D,Wang J,Zhou Z,He Z,Zhao Q
Fri Jan 01 00:00:00 UTC 2016
Novel Recombinant Protein FlaA N/C Protects against Radiation Injury via NF-κB Signaling.
Radiation research
Xu Y,Wu D,Fan Y,Li P,Du H,Shi J,Wang D,Zhou X
Fri Jan 01 00:00:00 UTC 2016
Novel Recombinant Protein FlaA N/C Protects against Radiation Injury via NF-κB Signaling.
Radiation research
Xu Y,Wu D,Fan Y,Li P,Du H,Shi J,Wang D,Zhou X
Fri Jan 01 00:00:00 UTC 2016
Carboxy-terminal modulator protein attenuated extracellular matrix deposit by inhibiting phospho-Akt, TGF-β1 and α-SMA in kidneys of diabetic mice.
Biochemical and biophysical research communications
Chen N,Hao J,Li L,Li F,Liu S,Duan H
Fri Jun 10 00:00:00 UTC 2016
Carboxy-terminal modulator protein attenuated extracellular matrix deposit by inhibiting phospho-Akt, TGF-β1 and α-SMA in kidneys of diabetic mice.
Biochemical and biophysical research communications
Chen N,Hao J,Li L,Li F,Liu S,Duan H
Fri Jun 10 00:00:00 UTC 2016
N-Methyl-D-aspartate Receptor Excessive Activation Inhibited Fetal Rat Lung Development In Vivo and In Vitro.
BioMed research international
Liao Z,Zhou X,Luo Z,Huo H,Wang M,Yu X,Cao C,Ding Y,Xiong Z,Yue S
Thu Feb 16 00:00:00 UTC 2017
N-Methyl-D-aspartate Receptor Excessive Activation Inhibited Fetal Rat Lung Development In Vivo and In Vitro.
BioMed research international
Liao Z,Zhou X,Luo Z,Huo H,Wang M,Yu X,Cao C,Ding Y,Xiong Z,Yue S
Thu Feb 16 00:00:00 UTC 2017
Calcium efflux from the endoplasmic reticulum regulates cisplatin-induced apoptosis in human cervical cancer HeLa cells.
Oncology letters
Shen L,Wen N,Xia M,Zhang YU,Liu W,Xu YE,Sun L
Fri Apr 01 00:00:00 UTC 2016
Calcium efflux from the endoplasmic reticulum regulates cisplatin-induced apoptosis in human cervical cancer HeLa cells.
Oncology letters
Shen L,Wen N,Xia M,Zhang YU,Liu W,Xu YE,Sun L
Fri Apr 01 00:00:00 UTC 2016
Establishment of a first-line second-line treatment model for human pulmonary adenocarcinoma.
Oncology letters
Wang L,Wang Y,Guan Q,Liu Y,He T,Wang J
Thu Dec 01 00:00:00 UTC 2016
Establishment of a first-line second-line treatment model for human pulmonary adenocarcinoma.
Oncology letters
Wang L,Wang Y,Guan Q,Liu Y,He T,Wang J
Thu Dec 01 00:00:00 UTC 2016
Development of a Biomimetic Chondroitin Sulfate-modified Hydrogel to Enhance the Metastasis of Tumor Cells.
Scientific reports
Liu Y,Wang S,Sun D,Liu Y,Liu Y,Wang Y,Liu C,Wu H,Lv Y,Ren Y,Guo X,Sun G,Ma X
Tue Jul 19 00:00:00 UTC 2016
Development of a Biomimetic Chondroitin Sulfate-modified Hydrogel to Enhance the Metastasis of Tumor Cells.
Scientific reports
Liu Y,Wang S,Sun D,Liu Y,Liu Y,Wang Y,Liu C,Wu H,Lv Y,Ren Y,Guo X,Sun G,Ma X
Tue Jul 19 00:00:00 UTC 2016
Quantitative Proteomic Profiling the Molecular Signatures of Annexin A5 in Lung Squamous Carcinoma Cells.
PloS one
Sun B,Bai Y,Zhang L,Gong L,Qi X,Li H,Wang F,Chi X,Jiang Y,Shao S
Wed Nov 20 00:00:00 UTC 2019
Quantitative Proteomic Profiling the Molecular Signatures of Annexin A5 in Lung Squamous Carcinoma Cells.
PloS one
Sun B,Bai Y,Zhang L,Gong L,Qi X,Li H,Wang F,Chi X,Jiang Y,Shao S
Wed Nov 20 00:00:00 UTC 2019
Quantitative proteomics reveals molecular mechanism of gamabufotalin and its potential inhibition on Hsp90 in lung cancer.
Oncotarget
Zhang L,Yu Z,Wang Y,Wang X,Zhang L,Wang C,Yue Q,Wang X,Deng S,Huo X,Tian X,Huang S,Zhang B,Ma X
Tue Nov 22 00:00:00 UTC 2016
Quantitative proteomics reveals molecular mechanism of gamabufotalin and its potential inhibition on Hsp90 in lung cancer.
Oncotarget
Zhang L,Yu Z,Wang Y,Wang X,Zhang L,Wang C,Yue Q,Wang X,Deng S,Huo X,Tian X,Huang S,Zhang B,Ma X
Tue Nov 22 00:00:00 UTC 2016
Targeting Dynamin 2 as a Novel Pathway to Inhibit Cardiomyocyte Apoptosis Following Oxidative Stress.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Gao D,Yang J,Wu Y,Wang Q,Wang Q,Lai EY,Zhu J
Mon Feb 06 00:00:00 UTC 2017
Targeting Dynamin 2 as a Novel Pathway to Inhibit Cardiomyocyte Apoptosis Following Oxidative Stress.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Gao D,Yang J,Wu Y,Wang Q,Wang Q,Lai EY,Zhu J
Mon Feb 06 00:00:00 UTC 2017
MiR-770-5p inhibits cisplatin chemoresistance in human ovarian cancer by targeting ERCC2.
Oncotarget
Zhao H,Yu X,Ding Y,Zhao J,Wang G,Wu X,Jiang J,Peng C,Guo GZ,Cui S
Tue Aug 16 00:00:00 UTC 2016
Gamabufotalin, a major derivative of bufadienolide, inhibits VEGF-induced angiogenesis by suppressing VEGFR-2 signaling pathway.
Oncotarget
Tang N,Shi L,Yu Z,Dong P,Wang C,Huo X,Zhang B,Huang S,Deng S,Liu K,Ma T,Wang X,Wu L,Ma XC
Tue Jan 19 00:00:00 UTC 2016
Gamabufotalin, a major derivative of bufadienolide, inhibits VEGF-induced angiogenesis by suppressing VEGFR-2 signaling pathway.
Oncotarget
Tang N,Shi L,Yu Z,Dong P,Wang C,Huo X,Zhang B,Huang S,Deng S,Liu K,Ma T,Wang X,Wu L,Ma XC
Tue Jan 19 00:00:00 UTC 2016
AMPK-mediated up-regulation of mTORC2 and MCL-1 compromises the anti-cancer effects of aspirin.
Oncotarget
Gao M,Kong Q,Hua H,Yin Y,Wang J,Luo T,Jiang Y
Tue Mar 29 00:00:00 UTC 2016
From the Cover: Tetrachlorobenzoquinone Exerts Neurological Proinflammatory Activity by Promoting HMGB1 Release, Which Induces TLR4 Clustering within the Lipid Raft.
Toxicological sciences : an official journal of the Society of Toxicology
Fu J,Shi Q,Song X,Liu Z,Wang Y,Wang Y,Song E,Song Y
Sat Oct 01 00:00:00 UTC 2016
MYBL2 guides autophagy suppressor VDAC2 in the developing ovary to inhibit autophagy through a complex of VDAC2-BECN1-BCL2L1 in mammals.
Autophagy
Yuan J,Zhang Y,Sheng Y,Fu X,Cheng H,Zhou R
Mon Apr 18 00:00:00 UTC 2016
Highly expressed UNC119 promotes hepatocellular carcinoma cell proliferation through Wnt/β-catenin signaling and predicts a poor prognosis.
American journal of cancer research
Lei B,Chai W,Wang Z,Liu R
Tue Dec 22 00:00:00 UTC 2015
Highly expressed UNC119 promotes hepatocellular carcinoma cell proliferation through Wnt/β-catenin signaling and predicts a poor prognosis.
American journal of cancer research
Lei B,Chai W,Wang Z,Liu R
Tue Dec 22 00:00:00 UTC 2015
SPAG6 silencing inhibits the growth of the malignant myeloid cell lines SKM-1 and K562 via activating p53 and caspase activation-dependent apoptosis.
International journal of oncology
Yang B,Wang L,Luo X,Chen L,Yang Z,Liu L
Sun Feb 01 00:00:00 UTC 2015
Olaquindox induces DNA damage via the lysosomal and mitochondrial pathway involving ROS production and p53 activation in HEK293 cells.
Environmental toxicology and pharmacology
Yang Y,Jiang L,She Y,Chen M,Li Q,Yang G,Geng C,Tang L,Zhong L,Jiang L,Liu X
Sun Nov 01 00:00:00 UTC 2015
Olaquindox induces DNA damage via the lysosomal and mitochondrial pathway involving ROS production and p53 activation in HEK293 cells.
Environmental toxicology and pharmacology
Yang Y,Jiang L,She Y,Chen M,Li Q,Yang G,Geng C,Tang L,Zhong L,Jiang L,Liu X
Sun Nov 01 00:00:00 UTC 2015
Artificial sweetener neohesperidin dihydrochalcone showed antioxidative, anti-inflammatory and anti-apoptosis effects against paraquat-induced liver injury in mice.
International immunopharmacology
Shi Q,Song X,Fu J,Su C,Xia X,Song E,Song Y
Tue Dec 01 00:00:00 UTC 2015
Artificial sweetener neohesperidin dihydrochalcone showed antioxidative, anti-inflammatory and anti-apoptosis effects against paraquat-induced liver injury in mice.
International immunopharmacology
Shi Q,Song X,Fu J,Su C,Xia X,Song E,Song Y
Tue Dec 01 00:00:00 UTC 2015
Tetrachlorobenzoquinone triggers the cleavage of Bid and promotes the cross-talk of extrinsic and intrinsic apoptotic signalings in pheochromocytoma (PC) 12 cells.
Neurotoxicology
Hu L,Su C,Song X,Shi Q,Fu J,Xia X,Xu D,Song E,Song Y
Wed Jul 01 00:00:00 UTC 2015
Tetrachlorobenzoquinone triggers the cleavage of Bid and promotes the cross-talk of extrinsic and intrinsic apoptotic signalings in pheochromocytoma (PC) 12 cells.
Neurotoxicology
Hu L,Su C,Song X,Shi Q,Fu J,Xia X,Xu D,Song E,Song Y
Wed Jul 01 00:00:00 UTC 2015
MicroRNA-206 attenuates tumor proliferation and migration involving the downregulation of NOTCH3 in colorectal cancer.
Oncology reports
Wang XW,Xi XQ,Wu J,Wan YY,Hui HX,Cao XF
Sun Mar 01 00:00:00 UTC 2015
Inhibition of migration and induction of apoptosis in LoVo human colon cancer cells by polysaccharides from Ganoderma lucidum.
Molecular medicine reports
Liang ZE,Yi YJ,Guo YT,Wang RC,Hu QL,Xiong XY
Sun Nov 01 00:00:00 UTC 2015
Inhibition of migration and induction of apoptosis in LoVo human colon cancer cells by polysaccharides from Ganoderma lucidum.
Molecular medicine reports
Liang ZE,Yi YJ,Guo YT,Wang RC,Hu QL,Xiong XY
Sun Nov 01 00:00:00 UTC 2015
Trophoblast glycoprotein promotes pancreatic ductal adenocarcinoma cell metastasis through Wnt/planar cell polarity signaling.
Molecular medicine reports
He P,Jiang S,Ma M,Wang Y,Li R,Fang F,Tian G,Zhang Z
Wed Jul 01 00:00:00 UTC 2015
Cannabinoid WIN55, 212-2 induces cell cycle arrest and inhibits the proliferation and migration of human BEL7402 hepatocellular carcinoma cells.
Molecular medicine reports
Xu D,Wang J,Zhou Z,He Z,Zhao Q
Tue Dec 01 00:00:00 UTC 2015
Cannabinoid WIN55, 212-2 induces cell cycle arrest and inhibits the proliferation and migration of human BEL7402 hepatocellular carcinoma cells.
Molecular medicine reports
Xu D,Wang J,Zhou Z,He Z,Zhao Q
Tue Dec 01 00:00:00 UTC 2015
Dynamic expression of miR-206-3p during mouse skin development is independent of keratinocyte differentiation.
Molecular medicine reports
Mu Y,Zhou H,Wu WJ,Hu LC,Chen HB
Tue Dec 01 00:00:00 UTC 2015
Liquiritigenin Protects Rats from Carbon Tetrachloride Induced Hepatic Injury through PGC-1α Pathway.
Evidence-based complementary and alternative medicine : eCAM
Zhang Y,He Y,Yu H,Ma F,Wu J,Zhang X
Wed Jul 22 00:00:00 UTC 2015
Liquiritigenin Protects Rats from Carbon Tetrachloride Induced Hepatic Injury through PGC-1α Pathway.
Evidence-based complementary and alternative medicine : eCAM
Zhang Y,He Y,Yu H,Ma F,Wu J,Zhang X
Wed Jul 22 00:00:00 UTC 2015
Postnatal alterations in GABAB receptor tone produce sensorimotor gating deficits and protein level differences in adulthood.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience
Bolton MM,Heaney CF,Murtishaw AS,Sabbagh JJ,Magcalas CM,Kinney JW
Wed Apr 01 00:00:00 UTC 2015
Postnatal alterations in GABAB receptor tone produce sensorimotor gating deficits and protein level differences in adulthood.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience
Bolton MM,Heaney CF,Murtishaw AS,Sabbagh JJ,Magcalas CM,Kinney JW
Wed Apr 01 00:00:00 UTC 2015
The safety and anti-hypercholesterolemic effect of coptisine in Syrian golden hamsters.
Lipids
He K,Ye X,Wu H,Wang Y,Zou Z,Ning N,Hu Y,Chen B,Fang X,Li X
Sun Feb 01 00:00:00 UTC 2015
The safety and anti-hypercholesterolemic effect of coptisine in Syrian golden hamsters.
Lipids
He K,Ye X,Wu H,Wang Y,Zou Z,Ning N,Hu Y,Chen B,Fang X,Li X
Sun Feb 01 00:00:00 UTC 2015
miR-140-5p regulates adipocyte differentiation by targeting transforming growth factor-β signaling.
Scientific reports
Zhang X,Chang A,Li Y,Gao Y,Wang H,Ma Z,Li X,Wang B
Fri Dec 11 00:00:00 UTC 2015
miR-140-5p regulates adipocyte differentiation by targeting transforming growth factor-β signaling.
Scientific reports
Zhang X,Chang A,Li Y,Gao Y,Wang H,Ma Z,Li X,Wang B
Fri Dec 11 00:00:00 UTC 2015
Panaxadiol Saponin and Dexamethasone Improve Renal Function in Lipopolysaccharide-Induced Mouse Model of Acute Kidney Injury.
PloS one
Chen Y,Du Y,Li Y,Wang X,Gao P,Yang G,Fang Y,Meng Y,Zhao X
Mon Apr 25 00:00:00 UTC 2016
A Critical Role for Cysteine 57 in the Biological Functions of Selenium Binding Protein-1.
International journal of molecular sciences
Ying Q,Ansong E,Diamond AM,Yang W
Wed Nov 18 00:00:00 UTC 2015
Study on the Dynamic Biological Characteristics of Sca-1(+) Hematopoietic Stem and Progenitor Cell Senescence.
Stem cells international
Geng S,Mu XY,Chen XB,Hou JY,Jia DY,Xu CY,Wang YP
Wed Jun 24 00:00:00 UTC 2015
FUS Interacts with HSP60 to Promote Mitochondrial Damage.
PLoS genetics
Deng J,Yang M,Chen Y,Chen X,Liu J,Sun S,Cheng H,Li Y,Bigio EH,Mesulam M,Xu Q,Du S,Fushimi K,Zhu L,Wu JY
Tue Sep 01 00:00:00 UTC 2015
FUS Interacts with HSP60 to Promote Mitochondrial Damage.
PLoS genetics
Deng J,Yang M,Chen Y,Chen X,Liu J,Sun S,Cheng H,Li Y,Bigio EH,Mesulam M,Xu Q,Du S,Fushimi K,Zhu L,Wu JY
Tue Sep 01 00:00:00 UTC 2015
Establishment and Characterization of Human Germline Stem Cell Line with Unlimited Proliferation Potentials and no Tumor Formation.
Scientific reports
Hou J,Niu M,Liu L,Zhu Z,Wang X,Sun M,Yuan Q,Yang S,Zeng W,Liu Y,Li Z,He Z
Fri Nov 20 00:00:00 UTC 2015
Establishment and Characterization of Human Germline Stem Cell Line with Unlimited Proliferation Potentials and no Tumor Formation.
Scientific reports
Hou J,Niu M,Liu L,Zhu Z,Wang X,Sun M,Yuan Q,Yang S,Zeng W,Liu Y,Li Z,He Z
Fri Nov 20 00:00:00 UTC 2015
HBx-related long non-coding RNA DBH-AS1 promotes cell proliferation and survival by activating MAPK signaling in hepatocellular carcinoma.
Oncotarget
Huang JL,Ren TY,Cao SW,Zheng SH,Hu XM,Hu YW,Lin L,Chen J,Zheng L,Wang Q
Tue Oct 20 00:00:00 UTC 2015
The Nucleocapsid Protein of Coronaviruses Acts as a Viral Suppressor of RNA Silencing in Mammalian Cells.
Journal of virology
Cui L,Wang H,Ji Y,Yang J,Xu S,Huang X,Wang Z,Qin L,Tien P,Zhou X,Guo D,Chen Y
Tue Sep 01 00:00:00 UTC 2015
Reactive stroma component COL6A1 is upregulated in castration-resistant prostate cancer and promotes tumor growth.
Oncotarget
Zhu YP,Wan FN,Shen YJ,Wang HK,Zhang GM,Ye DW
Wed Jun 10 00:00:00 UTC 2015
Reactive stroma component COL6A1 is upregulated in castration-resistant prostate cancer and promotes tumor growth.
Oncotarget
Zhu YP,Wan FN,Shen YJ,Wang HK,Zhang GM,Ye DW
Wed Jun 10 00:00:00 UTC 2015
Exposure to airborne PM2.5 suppresses microRNA expression and deregulates target oncogenes that cause neoplastic transformation in NIH3T3 cells.
Oncotarget
Liu C,Guo H,Cheng X,Shao M,Wu C,Wang S,Li H,Wei L,Gao Y,Tan W,Cheng S,Wu T,Yu D,Lin D
Tue Oct 06 00:00:00 UTC 2015
Exposure to airborne PM2.5 suppresses microRNA expression and deregulates target oncogenes that cause neoplastic transformation in NIH3T3 cells.
Oncotarget
Liu C,Guo H,Cheng X,Shao M,Wu C,Wang S,Li H,Wei L,Gao Y,Tan W,Cheng S,Wu T,Yu D,Lin D
Tue Oct 06 00:00:00 UTC 2015
Role of α-lipoic acid in LPS/d-GalN induced fulminant hepatic failure in mice: studies on oxidative stress, inflammation and apoptosis.
International immunopharmacology
Xia X,Su C,Fu J,Zhang P,Jiang X,Xu D,Hu L,Song E,Song Y
Wed Oct 01 00:00:00 UTC 2014
Role of α-lipoic acid in LPS/d-GalN induced fulminant hepatic failure in mice: studies on oxidative stress, inflammation and apoptosis.
International immunopharmacology
Xia X,Su C,Fu J,Zhang P,Jiang X,Xu D,Hu L,Song E,Song Y
Wed Oct 01 00:00:00 UTC 2014
Edwardsiella tarda-Induced cytotoxicity depends on its type III secretion system and flagellin.
Infection and immunity
Xie HX,Lu JF,Rolhion N,Holden DW,Nie P,Zhou Y,Yu XJ
Fri Aug 01 00:00:00 UTC 2014
Edwardsiella tarda-Induced cytotoxicity depends on its type III secretion system and flagellin.
Infection and immunity
Xie HX,Lu JF,Rolhion N,Holden DW,Nie P,Zhou Y,Yu XJ
Fri Aug 01 00:00:00 UTC 2014
RAD51B Activity and Cell Cycle Regulation in Response to DNA Damage in Breast Cancer Cell Lines.
Breast cancer : basic and clinical research
Lee PS,Fang J,Jessop L,Myers T,Raj P,Hu N,Wang C,Taylor PR,Wang J,Khan J,Jasin M,Chanock SJ
Tue Nov 04 00:00:00 UTC 2014
RAD51B Activity and Cell Cycle Regulation in Response to DNA Damage in Breast Cancer Cell Lines.
Breast cancer : basic and clinical research
Lee PS,Fang J,Jessop L,Myers T,Raj P,Hu N,Wang C,Taylor PR,Wang J,Khan J,Jasin M,Chanock SJ
Tue Nov 04 00:00:00 UTC 2014
Identification and validation of a novel microRNA-like molecule derived from a cytoplasmic RNA virus antigenome by bioinformatics and experimental approaches.
Virology journal
Shi J,Duan Z,Sun J,Wu M,Wang B,Zhang J,Wang H,Hu N,Hu Y
Tue Jul 01 00:00:00 UTC 2014
Identification and validation of a novel microRNA-like molecule derived from a cytoplasmic RNA virus antigenome by bioinformatics and experimental approaches.
Virology journal
Shi J,Duan Z,Sun J,Wu M,Wang B,Zhang J,Wang H,Hu N,Hu Y
Tue Jul 01 00:00:00 UTC 2014
Inhibition of 15-lipoxygenase (15-LOX) reverses hypoxia-induced down-regulation of potassium channels Kv1.5 and Kv2.1Inhibition of 15-lipoxygenase (15-LOX) reverses hypoxia-induced down-regulation of potassium channels Kv1.5 and Kv2.1.
International journal of clinical and experimental medicine
Liu W,Wang D,Song K,Chen L,Zhu Y,Liu P,Zhu Y
Wed Dec 31 00:00:00 UTC 2014
Inhibition of 15-lipoxygenase (15-LOX) reverses hypoxia-induced down-regulation of potassium channels Kv1.5 and Kv2.1Inhibition of 15-lipoxygenase (15-LOX) reverses hypoxia-induced down-regulation of potassium channels Kv1.5 and Kv2.1.
International journal of clinical and experimental medicine
Liu W,Wang D,Song K,Chen L,Zhu Y,Liu P,Zhu Y
Wed Dec 31 00:00:00 UTC 2014
Limb-girdle muscular dystrophy 1F is caused by a microdeletion in the transportin 3 gene.
Brain : a journal of neurology
Melià MJ,Kubota A,Ortolano S,Vílchez JJ,Gámez J,Tanji K,Bonilla E,Palenzuela L,Fernández-Cadenas I,Pristoupilová A,García-Arumí E,Andreu AL,Navarro C,Hirano M,Martí R
Wed May 01 00:00:00 UTC 2013
Limb-girdle muscular dystrophy 1F is caused by a microdeletion in the transportin 3 gene.
Brain : a journal of neurology
Melià MJ,Kubota A,Ortolano S,Vílchez JJ,Gámez J,Tanji K,Bonilla E,Palenzuela L,Fernández-Cadenas I,Pristoupilová A,García-Arumí E,Andreu AL,Navarro C,Hirano M,Martí R
Wed May 01 00:00:00 UTC 2013
The hepatitis B virus X protein downregulates NF-κB signaling pathways through decreasing the Notch signaling pathway in HBx-transformed L02 cells.
International journal of oncology
Luo J,Zhou H,Wang F,Xia X,Sun Q,Wang R,Cheng B
Wed May 01 00:00:00 UTC 2013
The hepatitis B virus X protein downregulates NF-κB signaling pathways through decreasing the Notch signaling pathway in HBx-transformed L02 cells.
International journal of oncology
Luo J,Zhou H,Wang F,Xia X,Sun Q,Wang R,Cheng B
Wed May 01 00:00:00 UTC 2013
Roles of 3,4-methylenedioxymethamphetamine (MDMA)-induced alteration of connexin43 and intracellular Ca(2+) oscillation in its cardiotoxicity.
Toxicology
Zhuo L,Liu Q,Liu L,Sun TY,Wang RS,Qu GQ,Liu Q,Liu Y,Ren L
Fri Aug 09 00:00:00 UTC 2013
Roles of 3,4-methylenedioxymethamphetamine (MDMA)-induced alteration of connexin43 and intracellular Ca(2+) oscillation in its cardiotoxicity.
Toxicology
Zhuo L,Liu Q,Liu L,Sun TY,Wang RS,Qu GQ,Liu Q,Liu Y,Ren L
Fri Aug 09 00:00:00 UTC 2013
Cytohesin-2 as a novel prognostic marker for hepatocellular carcinoma.
Oncology reports
Xu K,Gao J,Yang X,Yao Y,Liu Q
Sat Jun 01 00:00:00 UTC 2013
Cytohesin-2 as a novel prognostic marker for hepatocellular carcinoma.
Oncology reports
Xu K,Gao J,Yang X,Yao Y,Liu Q
Sat Jun 01 00:00:00 UTC 2013
Integrative genomics in combination with RNA interference identifies prognostic and functionally relevant gene targets for oral squamous cell carcinoma.
PLoS genetics
Xu C,Wang P,Liu Y,Zhang Y,Fan W,Upton MP,Lohavanichbutr P,Houck JR,Doody DR,Futran ND,Zhao LP,Schwartz SM,Chen C,Méndez E
Thu May 30 00:00:00 UTC 2013
Integrative genomics in combination with RNA interference identifies prognostic and functionally relevant gene targets for oral squamous cell carcinoma.
PLoS genetics
Xu C,Wang P,Liu Y,Zhang Y,Fan W,Upton MP,Lohavanichbutr P,Houck JR,Doody DR,Futran ND,Zhao LP,Schwartz SM,Chen C,Méndez E
Thu May 30 00:00:00 UTC 2013
Calpain mediates caspase-dependent apoptosis initiated by hydrogen peroxide in pancreatic acinar AR42J cells.
Free radical research
Weber H,Müller L,Jonas L,Schult C,Sparmann G,Schuff-Werner P
Wed May 01 00:00:00 UTC 2013
Calpain mediates caspase-dependent apoptosis initiated by hydrogen peroxide in pancreatic acinar AR42J cells.
Free radical research
Weber H,Müller L,Jonas L,Schult C,Sparmann G,Schuff-Werner P
Wed May 01 00:00:00 UTC 2013