Product References
ADP-ribose transferase PARP16 mediated-unfolded protein response contributes to neuronal cell damage in cerebral ischemia/reperfusion.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wang J,Xu J,Dong Y,Su Z,Su H,Cheng Q,Liu X
Wed Feb 01 00:00:00 UTC 2023
Dexmedetomidine attenuates airway inflammation and oxidative stress in asthma via the Nrf2 signaling pathway.
Molecular medicine reports
Xiao S,Zhou Y,Gao H,Yang D
Sun Jan 01 00:00:00 UTC 2023
Tumor necrosis factor-α reduces adiponectin production by decreasing transcriptional activity of peroxisome proliferator-activated receptor-γ in calf adipocytes.
Journal of dairy science
Yu H,Gao X,Ge Q,Tai W,Hao X,Shao Q,Fang Z,Chen M,Song Y,Gao W,Liu G,Du X,Li X
Sat Jul 01 00:00:00 UTC 2023
Electroacupuncture alleviates perioperative hypothalamus-pituitary-adrenal axis dysfunction via circRNA-miRNA-mRNA networks.
Frontiers in molecular neuroscience
Wang Y,Hu W,Han J,Zheng J,Jiang N,Feng Y,Tian Z
Sat Feb 11 00:00:00 UTC 2023
The tick saliva peptide HIDfsin2 promotes the tick-borne virus SFTSV replication in vitro by enhancing p38 signal pathway.
Archives of toxicology
Wang L,Sun F,Hu J,Zuo W,Zheng Y,Wu Y,Kwok HF,Cao Z
Thu Jun 01 00:00:00 UTC 2023
Sodium Butyrate Ameliorates Type 2 Diabetes-Related Sarcopenia Through IL-33-Independent ILC2s/IL-13/STAT3 Signaling Pathway.
Journal of inflammation research
Cao Y,Li Y,Han W,Jia X,Zhu P,Wei B,Cong X,Wang Z
Sat Feb 04 00:00:00 UTC 2023
TET1-TRPV4 Signaling Contributes to Bone Cancer Pain in Rats.
Brain sciences
Xu ZH,Niu Z,Liu Y,Liu PL,Lin XL,Zhang L,Chen L,Song Y,Sun R,Zhang HL
Mon Apr 10 00:00:00 UTC 2023
HBV X Protein Induces Degradation of UBXN7, a Novel Negative Regulator of NF-κB Signaling, to Promote HBV Replication.
Cellular and molecular gastroenterology and hepatology
Yuan S,Xu J,Wang M,Huang J,Ma S,Liu Y,Ke Y,Zeng X,Wu K,Wang J,Tian X,Zheng D,Yousaf T,Naz W,Sun J,Chen L,Guo D,Guo M,Sun G
Tue Dec 27 00:00:00 UTC 2022
A Novel Prenylflavonoid Icariside I Ameliorates Estrogen Deficiency-Induced Osteoporosis via Simultaneous Regulation of Osteoblast and Osteoclast Differentiation.
ACS pharmacology & translational science
Chen C,Wu M,Lei H,Cao Z,Wu F,Song Y,Zhang C,Qin M,Zhang C,Du R,Zhou J,Lu Y,Xie D,Zhang L
Fri Feb 10 00:00:00 UTC 2023
PKCθ Regulates Pituitary Adenoma Bone Invasion by Activating Osteoclast in NF-κB/IL-1β-Dependent Manner.
Cancers
Wang Q,Lei Z,Wang Z,Jiang Q,Zhang Z,Liu X,Xing B,Li S,Guo X,Liu Y,Li X,Shu K,Zhang H,Huang Y,Lei T
Mon Mar 06 00:00:00 UTC 2023
A conditional knockout rat resource of mitochondrial protein-coding genes via a DdCBE-induced premature stop codon.
Science advances
Tan L,Qi X,Kong W,Jin J,Lu D,Zhang X,Wang Y,Wang S,Dong W,Shi X,Chen W,Wang J,Li K,Xie Y,Gao L,Guan F,Gao K,Li C,Wang C,Hu Z,Zhang L,Guo X,Shen B,Ma Y
Fri Apr 14 00:00:00 UTC 2023
Exosomes derived from oral squamous cell carcinoma tissue accelerate diabetic wound healing.
American journal of physiology. Cell physiology
Zhang M,Guo J,Xiang K,Chen J,Wang C,Jiang T,Kang Y,Xu X,Li J,Yang X,Chen Z
Thu Jun 01 00:00:00 UTC 2023
Association between F-box-only protein 43 overexpression and hepatocellular carcinoma pathogenesis and prognosis.
Cancer medicine
Wu S,Qin L,Yang J,Wang J,Shen Y
Sat Apr 01 00:00:00 UTC 2023
Novel Benzo Five-Membered Heterocycle Derivatives as P-Glycoprotein Inhibitors: Design, Synthesis, Molecular Docking, and Anti-Multidrug Resistance Activity.
Journal of medicinal chemistry
Yang Z,Cai Y,Yang X,Li Y,Wu Q,Yu Y,Chen Z,Wei B,Tian JM,Bao X,Ye X,Chen J,Zhang H,Mou X,Sun X,Wang H
Thu Apr 27 00:00:00 UTC 2023
The protective role of EP300 in monocrotaline-induced pulmonary hypertension.
Frontiers in cardiovascular medicine
Yang L,Tian J,Wang J,Zeng J,Wang T,Lin B,Linneman J,Li L,Niu Y,Gou D,Zhang Y
Tue Mar 14 00:00:00 UTC 2023
TM6SF2 reduces lipid accumulation in vascular smooth muscle cells by inhibiting LOX-1 and CD36 expression.
Experimental cell research
Li TT,Cui YT,Li TH,Xiang Q,Chen YY,Zheng XL,Peng J,Tang ZH
Tue Aug 15 00:00:00 UTC 2023
Si-Ni-SAN ameliorates obesity through AKT/AMPK/HSL pathway-mediated lipolysis: Network pharmacology and experimental validation.
Journal of ethnopharmacology
Li J,Wu K,Zhong Y,Kuang J,Huang N,Guo X,Du H,Guo C,Li R,Zhu X,Zhang T,Gong L,Sheng L,Sun R
Fri Feb 10 00:00:00 UTC 2023
Decellularized brain extracellular matrix slice glioblastoma culture model recapitulates the interaction between cells and the extracellular matrix without a nutrient-oxygen gradient interference.
Acta biomaterialia
Wang C,Zhao Q,Zheng X,Li S,Chen J,Zhao H,Chen F,Cui L,Li W
Wed Mar 01 00:00:00 UTC 2023
Combined effects of EMP and RF field on emotional behavior in mice.
Frontiers in public health
Qin T,Liu L,Wang X,Guo L,Lin J,Du J,Xue Y,Lai P,Jing Y,Ding G
Tue Apr 04 00:00:00 UTC 2023
Baricitinib Attenuates Bleomycin-Induced Pulmonary Fibrosis in Mice by Inhibiting TGF-β1 Signaling Pathway.
Molecules (Basel, Switzerland)
Gu S,Liang J,Zhang J,Liu Z,Miao Y,Wei Y,Li S,Gu J,Cui Y,Xiao T,Li X,Yang C
Mon Feb 27 00:00:00 UTC 2023
LncRNA SPRY4-IT1 facilitates cell proliferation and angiogenesis of glioma via the miR-101-3p/EZH2/VEGFA signaling axis.
Cancer medicine
Wang J,Chen Y,Wang Q,Xu H,Jiang Q,Wang M,Li S,Chen Y,Wu C,Yu P,Xiao Z,Chen W,Lan Q
Wed Mar 01 00:00:00 UTC 2023
Collagen type ¿-loaded methacrylamide hyaluronic acid hydrogel microneedles alleviate stress urinary incontinence in mice: A novel treatment and prevention strategy.
Colloids and surfaces. B, Biointerfaces
Zhang S,Yang L,Hong S,Liu J,Cheng J,He Y,Hong L
Wed Feb 01 00:00:00 UTC 2023
Knockout of transient receptor potential ankyrin 1 (TRPA1) modulates the glial phenotype and alleviates perihematomal neuroinflammation after intracerebral hemorrhage in mice via MAPK/NF-κB signaling.
Neuroreport
Xia M,Chen YJ,Chen B,Ru X,Wang J,Lin J,Tang X,Chen W,Hu R,Li W,Feng H
Wed Feb 01 00:00:00 UTC 2023
Mechanism of YY1 mediating autophagy dependent ferroptosis in PM2.5 induced cardiac fibrosis.
Chemosphere
Hu H,Li L,Zhang H,Zhang Y,Liu Q,Chen M,Ning J,Pang Y,Hu W,Niu Y,Zhang R
Wed Feb 01 00:00:00 UTC 2023
Key domains and residues of the receptor MRGPRX1 recognizing the peptide ligand BAM8-22.
Peptides
Hu J,Wang L,Yang H,Meng Y,Tao M,Wu Y,Cao Z
Wed Mar 01 00:00:00 UTC 2023
Site-specific phosphorylation of tau impacts mitochondrial biology and response to stressors.
bioRxiv : the preprint server for biology
Isei MO,Girardi PA,Rodwell-Bullock J,Nehrke K,Johnson GV
Sun Feb 19 00:00:00 UTC 2023
Antidepressant-like effect of acute dose of Naringin involves suppression of NR1 and activation of protein kinase A/cyclic adenosine monophosphate response element-binding protein/brain-derived neurotrophic factor signaling in hippocampus.
Behavioural pharmacology
Wang G,Yang H,Zuo W,Mei X
Sat Apr 01 00:00:00 UTC 2023
Estrogen-mediated oar-miR-485-5p targets PPP1R13B to regulate myoblast proliferation in sheep.
International journal of biological macromolecules
Liu S,Liu Z,Wang P,Li W,Zhao S,Liu Y,Chu M
Mon May 01 00:00:00 UTC 2023
Hydrogen sulfide antagonizes formaldehyde-induced ferroptosis via preventing ferritinophagy by upregulation of GDF11 in HT22 cells.
Toxicology
Tang YH,Wu L,Huang HL,Zhang PP,Zou W,Tang XQ,Tang YY
Thu Jun 01 00:00:00 UTC 2023
Suppression of the gut microbiota-bile acid-FGF19 axis in patients with atrial fibrillation.
Cell proliferation
Zuo K,Fang C,Gao Y,Fu Y,Wang H,Li J,Zhong J,Yang X,Xu L
Wed Nov 01 00:00:00 UTC 2023
Artesunate, a new antimalarial clinical drug, exhibits potent anti-AML activity by targeting the ROS/Bim and TFRC/Fe2+ pathways.
British journal of pharmacology
Liu Y,Li H,Luo Z,Yu Y,Yang J,Zhang M,Law BYK,Huang Z,Li W
Wed Mar 01 00:00:00 UTC 2023
One Stone, Two Birds: A Peptide-Au(I) Infinite Coordination Supermolecule for the Confederate Physical and Biological Radiosensitization in Cancer Radiation Therapy.
Small (Weinheim an der Bergstrasse, Germany)
Wang J,Zhao J,Ma F,Gong L,Lu Y,Xiao W,Tang H,Gao C,Chen Y,Ma J,Gao Z,Yan J,Han S
Wed Mar 01 00:00:00 UTC 2023
VPS35 promotes cell proliferation via EGFR recycling and enhances EGFR inhibitors response in gastric cancer.
EBioMedicine
Yu J,Feng H,Sang Q,Li F,Chen M,Yu B,Xu Z,Pan T,Wu X,Hou J,Zhu Z,Yan C,Su L,Li J,Liu B
Wed Mar 01 00:00:00 UTC 2023
Peroxiredoxin-6 Released by Astrocytes Contributes to Neuroapoptosis During Ischemia.
Neuroscience
Hou JY,Zhou XL,Wang XY,Liang J,Xue Q
Tue Feb 21 00:00:00 UTC 2023
Targeting Chondroitin Sulphate Synthase 1 (Chsy1) Promotes Axon Growth Following Neurorrhaphy by Suppressing Versican Accumulation.
Molecules (Basel, Switzerland)
Liu CH,Ho YJ,Wang CY,Hsu CC,Chu YH,Hsu MY,Chen SJ,Hsiao WC,Liao WC
Wed Apr 26 00:00:00 UTC 2023
The distinct initiation sites and processing activities of TTLL4 and TTLL7 in glutamylation of brain tubulin.
The Journal of biological chemistry
Zhang X,Li X,Chen W,Wang Y,Diao L,Gao Y,Wang H,Bao L,Liang X,Wu HY
Sat Jul 01 00:00:00 UTC 2023
Deletion of Bak1 alleviates microglial necroptosis and neuroinflammation after experimental subarachnoid hemorrhage.
Journal of neurochemistry
Qiu X,Tao Q,Zhang L,Kuang C,Xie Y,Zhang L,Yin S,Peng J,Jiang Y
Wed Mar 01 00:00:00 UTC 2023
Anti-inflammatory and osteoprotective effects of Shi-Wei-Ru-Xiang pills on collagen-induced arthritis in rats via inhibiting MAPK and STAT3 pathways.
Journal of ethnopharmacology
Xiong H,Meng F,Luo M,Chen W,Tian J,Chen L,Ju Y,Mei Z
Tue Jan 10 00:00:00 UTC 2023
MRPL51 is a downstream target of FOXM1 in promoting the malignant behaviors of lung adenocarcinoma.
Oncology letters
Zhang W,Yu L,Xu C,Tang T,Cao J,Chen L,Pang X,Ren W
Sat Jul 01 00:00:00 UTC 2023
Transcription factor ETS proto-oncogene 1 contributes to neuropathic pain by regulating histone deacetylase 1 in primary afferent neurons.
Molecular pain
Zheng HL,Sun SY,Jin T,Zhang M,Zeng Y,Liu Q,Yang K,Wei R,Pan Z,Lin F
Sun Feb 12 00:00:00 UTC 2023
Transcription factor ETS proto-oncogene 1 contributes to neuropathic pain by regulating histone deacetylase 1 in primary afferent neurons.
Molecular pain
Zheng HL,Sun SY,Jin T,Zhang M,Zeng Y,Liu Q,Yang K,Wei R,Pan Z,Lin F
Sun Feb 12 00:00:00 UTC 2023
Jatrorrhizine Suppresses Murine-Norovirus-Triggered N-GSDMD-Dependent Pyroptosis in RAW264.7 Macrophages.
Vaccines
Fu M,Chen N,Zhou Y,Chen S,Xu W,Gong S,Geng L
Thu Jan 12 00:00:00 UTC 2023
MicroRNA‑30a‑5p regulates cypermethrin-induced apoptosis of Sertoli cells by targeting KLF9 in vitro.
Reproductive toxicology (Elmsford, N.Y.)
Wang Q,Xie JF,Yao TT,Wang XX,Guo QW,Wang LS,Yu Y,Xu LC
Tue Aug 01 00:00:00 UTC 2023
LncRNA SOCS2-AS1 promotes the progression of papillary thyroid cancer by destabilizing p53 protein.
Biochemical and biophysical research communications
Zhang X,Zhang X,Yang G,Wan L,Yin F,Li H,Yin D
Sun Aug 20 00:00:00 UTC 2023
Plasma exosomes induce inflammatory immune response in patients with acute myocardial infarction.
Archives of physiology and biochemistry
Gong Z,Wen M,Zhang W,Yu L,Huang C,Xu Y,Xia Z,Xu M,Xu J,Liang Q,Bao H,Cheng X
Sun Oct 01 00:00:00 UTC 2023
Polydopamine-Decorated PLCL Conduit to Induce Synergetic Effect of Electrical Stimulation and Topological Morphology for Peripheral Nerve Regeneration.
Small methods
Lu S,Chen W,Wang J,Guo Z,Xiao L,Wei L,Yu J,Yuan Y,Chen W,Bian M,Huang L,Liu Y,Zhang J,Li YL,Jiang LB
Wed Feb 01 00:00:00 UTC 2023
Magnify is a universal molecular anchoring strategy for expansion microscopy.
Nature biotechnology
Klimas A,Gallagher BR,Wijesekara P,Fekir S,DiBernardo EF,Cheng Z,Stolz DB,Cambi F,Watkins SC,Brody SL,Horani A,Barth AL,Moore CI,Ren X,Zhao Y
Thu Jun 01 00:00:00 UTC 2023
Proteomics Profiling Reveals the Molecular Signatures and Potential Therapeutic Targets of Human Nasopharyngeal Carcinoma.
Molecular & cellular proteomics : MCP
Fu Y,Liang X,Yang X,Liu J,Huang H,Zhang P,Li S,Zhu D,Zhang Y,Peng F,Chen Y,Chen Z
Thu Jun 01 00:00:00 UTC 2023
Stabilization of nuclear β-catenin by inhibiting KDM2A mediates cerebral ischemic tolerance.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zuo Y,Zhan L,Wen H,Xue J,Tan Y,Sun W,Xu E
Wed Mar 01 00:00:00 UTC 2023
Ginsenoside Rb1 improves energy metabolism after spinal cord injury.
Neural regeneration research
Wen S,Zou ZR,Cheng S,Guo H,Hu HS,Zeng FZ,Mei XF
Thu Jun 01 00:00:00 UTC 2023
CRISPR/Cas9 based gene editing of Frizzled class receptor 6 (FZD6) reveals its role in depressive symptoms through disrupting Wnt/β-catenin signaling pathway.
Journal of advanced research
Han H,Xu M,Wang J,Li MD,Yang Z
Wed Jun 14 00:00:00 UTC 2023
Pine pollen extract alleviates ethanol-induced oxidative stress and apoptosis in HepG2 cells via MAPK signaling.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Ma D,Wang Z,He Z,Wang Z,Chen Q,Qin F,Zeng M,Chen J
Sun Jan 01 00:00:00 UTC 2023
Neobavaisoflavone induces pyroptosis of liver cancer cells via Tom20 sensing the activated ROS signal.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li Y,Zhao R,Xiu Z,Yang X,Zhu Y,Han J,Li S,Li Y,Sun L,Li X,Jin N,Li Y
Tue Jul 25 00:00:00 UTC 2023
Network Pharmacology and Experimental Verification to Unveil the Mechanism of N-Methyl-D-Aspartic Acid Rescue Humantenirine-Induced Excitotoxicity.
Metabolites
Qi XJ,Huang CY,Zuo MT,Gong MD,Huang SJ,Tang MH,Liu ZY
Sat Jan 28 00:00:00 UTC 2023
Low-Dose Radiation Reduces Doxorubicin-Induced Myocardial Injury Through Mitochondrial Pathways.
Dose-response : a publication of International Hormesis Society
Zhao D,Jiang X,Meng X,Liu D,Du Y,Zhao L,Jiang H
Sat Feb 18 00:00:00 UTC 2023
Inhibition of deubiquitinase USP28 attenuates cyst growth in autosomal dominant polycystic kidney disease.
Biochemical pharmacology
Ren Y,Zhu X,Fu K,Zhang H,Zhao W,Lin Y,Fang Q,Wang J,Chen Y,Guo D
Sun Jan 01 00:00:00 UTC 2023
microRNA-627-5p inhibits colorectal cancer cell proliferation, migration and invasion by targeting Wnt2.
World journal of gastrointestinal oncology
Zhao DY,Yin TF,Sun XZ,Zhou YC,Wang QQ,Zhou GY,Yao SK
Wed Feb 15 00:00:00 UTC 2023
Bacterial infection promotes tumorigenesis of colorectal cancer via regulating CDC42 acetylation.
PLoS pathogens
Wang DN,Ni JJ,Li JH,Gao YQ,Ni FJ,Zhang ZZ,Fang JY,Lu J,Yao YF
Wed Feb 01 00:00:00 UTC 2023
METTL3 promotes SMSCs chondrogenic differentiation by targeting the MMP3, MMP13, and GATA3.
Regenerative therapy
Hu B,Zou X,Yu Y,Jiang Y,Xu H
Wed Mar 01 00:00:00 UTC 2023
Enhanced E6AP-mediated ubiquitination of ENO1 via LINC00663 contributes to radiosensitivity of breast cancer by regulating mitochondrial homeostasis.
Cancer letters
Ma J,Zhu J,Li J,Liu J,Kang X,Yu J
Fri Apr 28 00:00:00 UTC 2023
Prognostic values of Annexins and validation of the influence on cell proliferation, migration, and invasion in uveal melanoma.
American journal of translational research
Zhao A,Wang Z,Wang Y,Chen X
Tue Jun 13 00:00:00 UTC 2023
TIPE2 deletion improves the therapeutic potential of adoptively transferred NK cells.
Journal for immunotherapy of cancer
Bi J,Huang C,Jin X,Zheng C,Huang Y,Zheng X,Tian Z,Sun H
Wed Feb 01 00:00:00 UTC 2023
Inhibition of Hypothalamic FTO Activates STAT3 Signal through ERK1/2 Associated with Reductions in Food Intake and Body Weight.
Neuroendocrinology
Hu F,Yan HJ,Gao CX,Sun WW,Long YS
Fri Jan 13 00:00:00 UTC 2023
BACH1 deficiency prevents neointima formation and maintains the differentiated phenotype of vascular smooth muscle cells by regulating chromatin accessibility.
Nucleic acids research
Guo J,Qiu J,Jia M,Li Q,Wei X,Li L,Pan Q,Jin J,Ge F,Ma S,He Y,Lin J,Li Y,Ma J,Jiang N,Zhi X,Jiang L,Zhang J,Osto E,Jing Q,Wang X,Meng D
Mon May 22 00:00:00 UTC 2023
Acinous cell AR42J-derived exosome miR125b-5p promotes acute pancreatitis exacerbation by inhibiting M2 macrophage polarization via PI3K/AKT signaling pathway.
World journal of gastrointestinal surgery
Zheng Z,Cao F,Ding YX,Lu JD,Fu YQ,Liu L,Guo YL,Liu S,Sun HC,Cui YQ,Li F
Thu Apr 27 00:00:00 UTC 2023
Dual-specificity phosphatase 26-dificient neurons are susceptible to oxygen-glucose deprivation/reoxygenation-evoked apoptosis and proinflammatory response by affecting the TAK1-medaited JNK/P38 MAPK pathway.
International immunopharmacology
Zhao H,Lv J,Meng L,Lv J,Li Z
Sat Apr 01 00:00:00 UTC 2023
Hydrogen sulfide alleviates mitochondrial damage and ferroptosis by regulating OPA3-NFS1 axis in doxorubicin-induced cardiotoxicity.
Cellular signalling
Wang Y,Ying X,Wang Y,Zou Z,Yuan A,Xiao Z,Geng N,Qiao Z,Li W,Lu X,Pu J
Sat Jul 01 00:00:00 UTC 2023
IGFBP3 induced by the TGF-β/EGFRvIII transactivation contributes to the malignant phenotype of glioblastoma.
iScience
Zhang X,Wang G,Gong Y,Zhao L,Song P,Zhang H,Zhang Y,Ju H,Wang X,Wang B,Ren H,Zhu X,Dong Y
Fri May 19 00:00:00 UTC 2023
Grouper cGAS is a negative regulator of STING-mediated interferon response.
Frontiers in immunology
Zhang L,Zhang X,Liao J,Xu L,Kang S,Chen H,Sun M,Wu S,Xu Z,Wei S,Qin Q,Wei J
Tue Feb 28 00:00:00 UTC 2023
Zinc Promotes Spinal Cord Injury Recovery by Blocking the Activation of NLRP3 Inflammasome Through SIRT3-Mediated Autophagy.
Neurochemical research
Xu C,Zhou Z,Zhao H,Lin S,Zhang P,Tian H,Mei X
Wed Feb 01 00:00:00 UTC 2023
Hyperactivation of TRPV4 causes the hippocampal pyroptosis pathway and results in cognitive impairment in LPS-treated mice.
Behavioural brain research
Guo D,Xu Y,Wang Y,Zhong X,Liu Z,Li S,Xu X,Zhang J,Xiong T,Cao W,Liang J
Wed Feb 15 00:00:00 UTC 2023
MUC1 promotes glioblastoma progression and TMZ resistance by stabilizing EGFRvIII.
Pharmacological research
Tong F,Zhao JX,Fang ZY,Cui XT,Su DY,Liu X,Zhou JH,Wang GX,Qiu ZJ,Liu SZ,Fu JQ,Kang CS,Wang JC,Wang QX
Sun Jan 01 00:00:00 UTC 2023
UBE2E2 enhances Snail-mediated epithelial-mesenchymal transition and Nrf2-mediated antioxidant activity in ovarian cancer.
Cell death & disease
Hong X,Ma N,Li D,Zhang M,Dong W,Huang J,Ci X,Zhang S
Fri Feb 10 00:00:00 UTC 2023
UBE2E2 enhances Snail-mediated epithelial-mesenchymal transition and Nrf2-mediated antioxidant activity in ovarian cancer.
Cell death & disease
Hong X,Ma N,Li D,Zhang M,Dong W,Huang J,Ci X,Zhang S
Fri Feb 10 00:00:00 UTC 2023
Amyloid futures in the expanding pathology of brain aging and dementia.
Alzheimer's & dementia : the journal of the Alzheimer's Association
Thorwald MA,Silva J,Head E,Finch CE
Thu Jun 01 00:00:00 UTC 2023
Endothelial progenitor cell-derived exosomes promote anti-inflammatory macrophages via SOCS3/JAK2/STAT3 axis and improve the outcome of spinal cord injury.
Journal of neuroinflammation
Yuan F,Peng W,Yang Y,Xu J,Liu Y,Xie Y,Huang T,Shi C,Ding Y,Li C,Qin T,Xie S,Zhu F,Lu H,Huang J,Hu J
Fri Jun 30 00:00:00 UTC 2023
Application of a JEG-3 organoid model to study HLA-G function in the trophoblast.
Frontiers in immunology
Zhuang BM,Cao DD,Liu XF,Wang L,Lin XL,Duan YG,Lee CL,Chiu PCN,Yeung WSB,Yao YQ
Tue Apr 04 00:00:00 UTC 2023
Deletion of transglutaminase 2 from astrocytes significantly improves their ability to promote neurite outgrowth on an inhibitory matrix.
bioRxiv : the preprint server for biology
Emerson J,Delgado T,Girardi P,Johnson GV
Mon Feb 06 00:00:00 UTC 2023
Discovery of a natural small-molecule AMP-activated kinase activator that alleviates nonalcoholic steatohepatitis.
Marine life science & technology
Chen J,Xu L,Zhang XQ,Liu X,Zhang ZX,Zhu QM,Liu JY,Iqbal MO,Ding N,Shao CL,Wei MY,Gu YC
Mon May 01 00:00:00 UTC 2023
Resveratrol-loaded macrophage exosomes alleviate multiple sclerosis through targeting microglia.
Journal of controlled release : official journal of the Controlled Release Society
Zheng X,Sun K,Liu Y,Yin X,Zhu H,Yu F,Zhao W
Sun Jan 01 00:00:00 UTC 2023
SIRT1 attenuates blood-spinal cord barrier disruption after spinal cord injury by deacetylating p66Shc.
Redox biology
Jiang T,Qin T,Gao P,Tao Z,Wang X,Wu M,Gu J,Chu B,Zheng Z,Yi J,Xu T,Huang Y,Liu H,Zhao S,Ren Y,Chen J,Yin G
Sat Apr 01 00:00:00 UTC 2023
Aβ Induces Neuroinflammation and Microglial M1 Polarization via cGAS-STING-IFITM3 Signaling Pathway in BV-2 Cells.
Neurochemical research
Wu Z,Tang W,Ibrahim FEEM,Chen X,Yan H,Tao C,Wang Z,Guo Y,Fu Y,Wang Q,Ge Y
Fri Sep 01 00:00:00 UTC 2023
GNB1 promotes hepatocellular carcinoma progression by targeting BAG2 to activate P38/MAPK signaling.
Cancer science
Zhang X,Dong K,Zhang J,Kuang T,Luo Y,Yu J,Yu J,Wang W
Mon May 01 00:00:00 UTC 2023
Chromatin remodeling and nucleoskeleton synergistically control osteogenic differentiation in different matrix stiffnesses.
Materials today. Bio
Xu X,Zhang H,Li Y,Liu F,Jing Z,Ren M,Chen T,Fu Y,Wu Y,Ji P,Yang S
Thu Jun 01 00:00:00 UTC 2023
Sevoflurane induces neurotoxic effects on developing neurons through the WNK1/NKCC1/Ca2+ /Drp-1 signalling pathway.
Clinical and experimental pharmacology & physiology
Bai YF,Li WJ,Ji YW,An LX,Zhang L,Li JF
Mon May 01 00:00:00 UTC 2023
Zcf24, a zinc-finger transcription factor, is required for lactate catabolism and inhibits commensalism in Candida albicans.
Molecular microbiology
Sun Q,Dong B,Yang D,Yu J,Ren T,Wang T,Yang L,Lu Y,Su C
Sun Jan 01 00:00:00 UTC 2023
Arctiin-encapsulated DSPE-PEG bubble-like nanoparticles inhibit alveolar epithelial type 2 cell senescence to alleviate pulmonary fibrosis via the p38/p53/p21 pathway.
Frontiers in pharmacology
Xiong D,Gao F,Shao J,Pan Y,Wang S,Wei D,Ye S,Chen Y,Chen R,Yue B,Li J,Chen J
Sat Apr 01 00:00:00 UTC 2023
Mutation of the attractin gene impairs working memory in rats.
Brain and behavior
Liu MQ,Xue C,Li XH,Ding HQ,Zhang MY,Chen K,Li Y,Gao SZ,Xu XJ,Zhang WN
Wed Feb 01 00:00:00 UTC 2023
Simiao San alleviates hyperuricemia and kidney inflammation by inhibiting NLRP3 inflammasome and JAK2/STAT3 signaling in hyperuricemia mice.
Journal of ethnopharmacology
Zhang Y,Wang S,Dai X,Liu T,Liu Y,Shi H,Yin J,Xu T,Zhang Y,Zhao D,Sukhorukov V,Orekhov AN,Gao S,Wang L,Zhang D
Thu Aug 10 00:00:00 UTC 2023
Effects of Spatial Expression of Activating Transcription Factor 4 on the Pathogenicity of Two Phenotypes of Bovine Viral Diarrhea Virus by Regulating the Endoplasmic Reticulum-Mediated Autophagy Process.
Microbiology spectrum
Wang J,Chen KY,Wang SH,Liu Y,Zhao YQ,Yang L,Yang GH,Wang XJ,Zhu YH,Yin JH,Wang JF
Mon Mar 20 00:00:00 UTC 2023
S100 Calcium Binding Protein A16 Promotes Cell Proliferation by triggering LATS1 ubiquitin degradation mediated by CUL4A ligase to inhibit Hippo pathway in Glioma development.
International journal of biological sciences
Hu Y,Zhang R,Lu S,Zhang W,Wang D,Ge Y,Jiang F,Qin X,Liu Y
Mon May 15 00:00:00 UTC 2023
TANGO1 interacts with NRTN to promote hepatocellular carcinoma progression by regulating the PI3K/AKT/mTOR signaling pathway.
Biochemical pharmacology
Man J,Zhou W,Zuo S,Zhao X,Wang Q,Ma H,Li HY
Sat Jul 01 00:00:00 UTC 2023
The overexpression of actin related protein 2/3 complex subunit 1B(ARPC1B) promotes the ovarian cancer progression via activation of the Wnt/β-catenin signaling pathway.
Frontiers in immunology
Huang J,Zhou H,Tan C,Mo S,Liu T,Kuang Y
Tue Jun 13 00:00:00 UTC 2023
Revealing active components and action mechanism of Fritillariae Bulbus against non-small cell lung cancer through spectrum-effect relationship and proteomics.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu FJ,Chen XY,Yang J,Zhao Z,Wang QL,Li P,Jiang Y,Li HJ
Wed Feb 01 00:00:00 UTC 2023
Low rumen degradable starch promotes the growth performance of goats by increasing protein synthesis in skeletal muscle via the AMPK-mTOR pathway.
Animal nutrition (Zhongguo xu mu shou yi xue hui)
Liang Z,Jin C,Bai H,Liang G,Su X,Wang D,Yao J
Thu Jun 01 00:00:00 UTC 2023
PDCD6 Promotes Hepatocellular Carcinoma Cell Proliferation and Metastasis through the AKT/GSK3β/β-catenin Pathway.
Biomedical and environmental sciences : BES
Wen SY,Liu YT,Wei BY,Ma JQ,Chen YY
Mon Mar 20 00:00:00 UTC 2023
Differential effects of WRAP53 transcript variants on non-small cell lung cancer cell behaviors.
PloS one
Zhu Y,Sun W,Jiang X,Bai R,Luo Y,Gao Y,Li S,Huang Z,Gong Y,Xie C
Fri Feb 03 00:00:00 UTC 2023
Systemic treatment with GnRH agonist produces antidepressant-like effects in LPS induced depression male mouse model.
Neuropharmacology
Xu Y,Li W,Shi M,Xu X,Guo D,Liu Z,Chen L,Zhong X,Cao W
Tue Aug 01 00:00:00 UTC 2023
Tibetan medicine Si-Wei-Qiang-Wei Powder ameliorates cholecystitis via inhibiting the production of pro-inflammatory cytokines and regulating the MAPK signaling pathway.
Journal of ethnopharmacology
Zheng Z,Xiong H,Zhao Z,Zhou K,Fu M,Liu X,Mei Z
Wed Mar 01 00:00:00 UTC 2023
Anisomycin inhibits Coxsackievirus B replication by promoting the lysosomal degradation of eEF1A1.
Antiviral research
Shao E,Zhao S,Dong Y,Wang Y,Fei Y,Li S,Wang L,Bashir T,Luan T,Lin L,Wang Y,Zhao W,Zhong Z
Sat Jul 01 00:00:00 UTC 2023
Docosahexaenoic Acid Alters Lipid Metabolism Processes via H3K9ac Epigenetic Modification in Dairy Goat.
Journal of agricultural and food chemistry
Wu J,Luo J,He Q,Xia Y,Tian H,Zhu L,Li C,Loor JJ
Wed Jun 07 00:00:00 UTC 2023
Design, synthesis and biological evaluation of novel phenylfuran-bisamide derivatives as P-glycoprotein inhibitors against multidrug resistance in MCF-7/ADR cell.
European journal of medicinal chemistry
Yang Z,Yang X,Li Y,Cai Y,Yu Y,Zhuang W,Sun X,Li Q,Bao X,Ye X,Tian J,Wei B,Chen J,Wu Q,Zhang H,Mou X,Wang H
Wed Feb 15 00:00:00 UTC 2023
Chlorogenic Acid Alleviates Hepatic Ischemia-Reperfusion Injury by Inhibiting Oxidative Stress, Inflammation, and Mitochondria-Mediated Apoptosis In Vivo and In Vitro.
Inflammation
Li K,Feng Z,Wang L,Ma X,Wang L,Liu K,Geng X,Peng C
Thu Jun 01 00:00:00 UTC 2023
The dichloromethane extract of Callicarpa longissima rich in diterpenoid phenols exerts anti-inflammatory effect via inhibiting the TLR4/NF-κB signaling pathway.
Journal of ethnopharmacology
Li LC,Fu YX,Ning DS,Yu LL,Zou ZQ,Pan ZH
Thu Apr 06 00:00:00 UTC 2023
SECTM1 is upregulated in immuno-hot tumors and predicts immunotherapeutic efficacy in multiple cancers.
iScience
Mei J,Fu Z,Cai Y,Song C,Zhou J,Zhu Y,Mao W,Xu J,Yin Y
Fri Feb 17 00:00:00 UTC 2023
PDLIM3 supports hedgehog signaling in medulloblastoma by facilitating cilia formation.
Cell death and differentiation
Zhang J,Yang Y,Li X,Li G,Mizukami T,Liu Y,Wang Y,Xu G,Roder H,Zhang L,Yang ZJ
Mon May 01 00:00:00 UTC 2023
ORP8 inhibits renal cell carcinoma progression by accelerating Stathmin1 degradation and microtubule polymerization.
Experimental cell research
Zhang L,Pan Q,Wu Y,Zhang P,Li S,Xu Y,Li D,Zheng M,Pei D,Wang Q
Thu Jun 01 00:00:00 UTC 2023
Piezo1 suppression reduces demyelination after intracerebral hemorrhage.
Neural regeneration research
Qu J,Zong HF,Shan Y,Zhang SC,Guan WP,Yang Y,Zhao HL
Tue Aug 01 00:00:00 UTC 2023
CB2R activation ameliorates late adolescent chronic alcohol exposure-induced anxiety-like behaviors during withdrawal by preventing morphological changes and suppressing NLRP3 inflammasome activation in prefrontal cortex microglia in mice.
Brain, behavior, and immunity
Li J,Wang H,Liu D,Li X,He L,Pan J,Shen Q,Peng Y
Mon May 01 00:00:00 UTC 2023
Marine-Derived Natural Product HDYL-GQQ-495 Targets P62 to Inhibit Autophagy.
Marine drugs
Li Q,Fan J,Chen Y,Liu Y,Liu H,Jiang W,Li D,Dang Y
Fri Jan 20 00:00:00 UTC 2023
Mechanism Repositioning Based on Integrative Pharmacology: Anti-Inflammatory Effect of Safflower in Myocardial Ischemia-Reperfusion Injury.
International journal of molecular sciences
Zhao F,Jiang H,Zhang T,Chen H,Li W,Li X,Wang P,Xu H
Fri Mar 10 00:00:00 UTC 2023
Targeting plasmodium α-tubulin-1 to block malaria transmission to mosquitoes.
Frontiers in cellular and infection microbiology
Zhang G,Niu G,Hooker-Romera D,Shabani S,Ramelow J,Wang X,Butler NS,James AA,Li J
Tue Apr 04 00:00:00 UTC 2023
IGF2 inhibits hippocampal over-activated microglia and alleviates depression-like behavior in LPS- treated male mice.
Brain research bulletin
Guo D,Xu Y,Liu Z,Wang Y,Xu X,Li C,Li S,Zhang J,Xiong T,Cao W,Liang J
Wed Mar 01 00:00:00 UTC 2023
ME2 Promotes Hepatocellular Carcinoma Cell Migration through Pyruvate.
Metabolites
Yang Y,Zhang Z,Li W,Li L,Zhou Y,Du W
Mon Apr 10 00:00:00 UTC 2023
Resveratrol Modulates the Redox Response and Bile Acid Metabolism to Maintain the Cholesterol Homeostasis in Fish Megalobrama amblycephala Offered a High-Carbohydrate Diet.
Antioxidants (Basel, Switzerland)
Ge Y,Zhang L,Chen W,Sun M,Liu W,Li X
Tue Jan 03 00:00:00 UTC 2023
Deletion of Transglutaminase 2 from Mouse Astrocytes Significantly Improves Their Ability to Promote Neurite Outgrowth on an Inhibitory Matrix.
International journal of molecular sciences
Emerson J,Delgado T,Girardi P,Johnson GVW
Thu Mar 23 00:00:00 UTC 2023
Deletion of Transglutaminase 2 from Mouse Astrocytes Significantly Improves Their Ability to Promote Neurite Outgrowth on an Inhibitory Matrix.
International journal of molecular sciences
Emerson J,Delgado T,Girardi P,Johnson GVW
Thu Mar 23 00:00:00 UTC 2023
In silico, in vitro and in vivo studies: Dibutyl phthalate promotes prostate cancer cell proliferation by activating Forkhead Box M1 and remission after Natura-α pretreatment.
Toxicology
Bu H,Tang S,Liu G,Miao C,Zhou X,Yang H,Liu B
Sat Apr 01 00:00:00 UTC 2023
PGC-1α Affects Epileptic Seizures by Regulating Mitochondrial Fusion in Epileptic Rats.
Neurochemical research
Li D,Zhang L,Tuo J,Zhang F,Tai Z,Liu X,Qiu X,Zhang H,Yang J,Wang J,Luo Z,Xu Z
Mon May 01 00:00:00 UTC 2023
Scarless wound healing programmed by core-shell microneedles.
Nature communications
Zhang Y,Wang S,Yang Y,Zhao S,You J,Wang J,Cai J,Wang H,Wang J,Zhang W,Yu J,Han C,Zhang Y,Gu Z
Sat Jun 10 00:00:00 UTC 2023
3, 14, 19-Triacetyl Andrographolide alleviates the cognitive dysfunction of 3 × Tg-AD mice by inducing initiation and promoting degradation process of autophagy.
Phytotherapy research : PTR
Zhang H,Xie X,Shi J,Zhao Q,Luo D,Hao J,Zhou Y,Han G,Pang X
Wed Feb 01 00:00:00 UTC 2023
ADAMTS7-Mediated Complement Factor H Degradation Potentiates Complement Activation to Contributing to Renal Injuries.
Journal of the American Society of Nephrology : JASN
Ma Z,Mao C,Jia Y,Yu F,Xu P,Tan Y,Zou QH,Zhou XJ,Kong W,Fu Y
Wed Feb 01 00:00:00 UTC 2023
1,2,4,5-Tetrazine-tethered probes for fluorogenically imaging superoxide in live cells with ultrahigh specificity.
Nature communications
Jiang X,Li M,Wang Y,Wang C,Wang Y,Shen T,Shen L,Liu X,Wang Y,Li X
Tue Mar 14 00:00:00 UTC 2023
PPARα is involved in high-fat diet-induced risk avoidance impairment via the regulation of hippocampal BDNF.
Food & function
Zhu XY,Luo J,Wei XJ,Pan LH,Xu XF,Hu LB,Dong YB,Jiang YK,Dong FY,Xu L,Xu LM,Liu AM,Zhang XQ
Mon Jul 17 00:00:00 UTC 2023
ROS-responsive polymer nanoparticles with enhanced loading of dexamethasone effectively modulate the lung injury microenvironment.
Acta biomaterialia
Muhammad W,Zhu J,Zhai Z,Xie J,Zhou J,Feng X,Feng B,Pan Q,Li S,Venkatesan R,Li P,Cao H,Gao C
Mon Aug 01 00:00:00 UTC 2022
Bioglass promotes wound healing by inhibiting endothelial cell pyroptosis through regulation of the connexin 43/reactive oxygen species (ROS) signaling pathway.
Laboratory investigation; a journal of technical methods and pathology
Zhang K,Chai B,Ji H,Chen L,Ma Y,Zhu L,Xu J,Wu Y,Lan Y,Li H,Feng Z,Xiao J,Zhang H,Xu K
Sat Jan 01 00:00:00 UTC 2022
Inhibiting Sphingosine 1-Phosphate Receptor Subtype 3 Attenuates Brain Damage During Ischemia-Reperfusion Injury by Regulating nNOS/NO and Oxidative Stress.
Frontiers in neuroscience
Fan X,Chen H,Xu C,Wang Y,Yin P,Li M,Tang Z,Jiang F,Wei W,Song J,Li G,Zhong D
Mon May 02 00:00:00 UTC 2022
RFC2: a prognosis biomarker correlated with the immune signature in diffuse lower-grade gliomas.
Scientific reports
Zhao X,Wang Y,Li J,Qu F,Fu X,Liu S,Wang X,Xie Y,Zhang X
Thu Feb 24 00:00:00 UTC 2022
IL-37 isoform D acts as an inhibitor of soluble ST2 to boost type 2 immune homeostasis in white adipose tissue.
Cell death discovery
Li C,Zhao M,Zhao M,Chen N,Guo Y,Du Y,Zhang Y,Cao B,Zhan B,Guo C,Li Y,Li Y,Shi Y,Zhu F,Zhang L,Wang Q
Tue Apr 05 00:00:00 UTC 2022
mTOR inhibition attenuates chemosensitivity through the induction of chemotherapy resistant persisters.
Nature communications
Liu Y,Azizian NG,Sullivan DK,Li Y
Thu Nov 17 00:00:00 UTC 2022
Difenoconazole disrupts the blood-brain barrier and results in neurotoxicity in carp by inhibiting the Nrf2 pathway mediated ROS accumulation.
Ecotoxicology and environmental safety
Liu F,Wang Y,Chen L,Bello BK,Zhang T,Yang H,Li X,Pan E,Feng H,Dong J
Sat Oct 01 00:00:00 UTC 2022
Acid ceramidase targeting pyruvate kinase affected trypsinogen activation in acute pancreatitis.
Molecular medicine (Cambridge, Mass.)
Xiao J,Zeng W,Zhang P,Zhou Y,Fang Q
Tue Sep 06 00:00:00 UTC 2022
p62 Promotes the Mitochondrial Localization of p53 through Its UBA Domain and Participates in Regulating the Sensitivity of Ovarian Cancer Cells to Cisplatin.
International journal of molecular sciences
Kong Q,Yan X,Cheng M,Jiang X,Xu L,Shen L,Yu H,Sun L
Fri Mar 18 00:00:00 UTC 2022
Transcriptome study digs out BMP2 involved in adipogenesis in sheep tails.
BMC genomics
Jin M,Fei X,Li T,Lu Z,Chu M,Di R,He X,Wang X,Wei C
Tue Jun 21 00:00:00 UTC 2022
MARCH1 promotes the growth and maintaining of stem cell-like characteristics of gastric cancer cells by activating the Wnt/β-catenin signaling pathway.
Tissue & cell
Liang W,Zhang T,Huo J,Yang J
Sat Oct 01 00:00:00 UTC 2022
Vagus nerve stimulation promotes the M1-to-M2 transition via inhibition of TLR4/NF-κB in microglial to rescue the reperfusion injury.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association
Zhang L,Zhang X,Liu Y,Wang S,Jia G
Thu Sep 01 00:00:00 UTC 2022
Supramolecular Hydrogel-Wrapped Gingival Mesenchymal Stem Cells in Cutaneous Radiation Injury.
Cells
Nie S,Ren C,Liang X,Cai H,Sun H,Liu F,Ji K,Wang Y,Liu Q
Fri Sep 30 00:00:00 UTC 2022
Oxidative stress from DGAT1 oncoprotein inhibition in melanoma suppresses tumor growth when ROS defenses are also breached.
Cell reports
Wilcock DJ,Badrock AP,Wong CW,Owen R,Guerin M,Southam AD,Johnston H,Telfer BA,Fullwood P,Watson J,Ferguson H,Ferguson J,Lloyd GR,Jankevics A,Dunn WB,Wellbrock C,Lorigan P,Ceol C,Francavilla C,Smith MP,Hurlstone AFL
Tue Jun 21 00:00:00 UTC 2022
A homozygous loss-of-function mutation in FBXO43 causes human non-obstructive azoospermia.
Clinical genetics
Wu H,Zhang X,Shen Q,Liu Y,Gao Y,Wang G,Lv M,Hua R,Xu Y,Zhou P,Wei Z,Tao F,He X,Cao Y,Liu M
Sat Jan 01 00:00:00 UTC 2022
Activation of Transcription Factor EB Is Associated With Adipose Tissue Lipolysis in Dairy Cows With Subclinical Ketosis.
Frontiers in veterinary science
Yu H,Gao X,Loor JJ,Jiang Q,Fang Z,Hao X,Shi Z,Fan M,Chen M,Li X,Liu G,Wang Z,Li X,Du X
Sat Feb 26 00:00:00 UTC 2022
Rutin-activated adipose tissue thermogenesis is correlated with increased intestinal short-chain fatty acid levels.
Phytotherapy research : PTR
Cheng L,Shi L,He C,Wang C,Lv Y,Li H,An Y,Dai H,Duan Y,Zhang H,Huang Y,Fu W,Meng Y,Zhao B
Wed Jun 01 00:00:00 UTC 2022
Knockdown of CLAUDIN-6 Inhibited Apoptosis and Induced Proliferation of Bovine Cumulus Cells.
International journal of molecular sciences
Wang W,Zou H,Chen N,Tian Y,Hao H,Pang Y,Zhao X,Zhu H,Yu D,Du W
Sun Oct 30 00:00:00 UTC 2022
Morphological and Proteomic Analyses of Soybean Seedling Interaction Mechanism Affected by Fiber Crosslinked with Zinc-Oxide Nanoparticles.
International journal of molecular sciences
Komatsu S,Murata K,Yakeishi S,Shimada K,Yamaguchi H,Hitachi K,Tsuchida K,Obi R,Akita S,Fukuda R
Sun Jul 03 00:00:00 UTC 2022
Mesenchymal stem cells protect against ferroptosis via exosome-mediated stabilization of SLC7A11 in acute liver injury.
Cell death & disease
Lin F,Chen W,Zhou J,Zhu J,Yao Q,Feng B,Feng X,Shi X,Pan Q,Yu J,Li L,Cao H
Sat Mar 26 00:00:00 UTC 2022
X-box binding protein 1: A new metabolic mediator and drug target of metformin?
Frontiers in pharmacology
Lou K,Sun P,Zhang C,Jiang Q,Pang S
Tue Nov 29 00:00:00 UTC 2022
Silica nanoparticle exposure inhibits surfactant protein A and B in A549 cells through ROS-mediated JNK/c-Jun signaling pathway.
Environmental toxicology
Yang X,Yang P,Zhang J,Yang Y,Xiong M,Shi F,Li N,Jin Y
Thu Sep 01 00:00:00 UTC 2022
Elevated phosphorylation of EGFR in NSCLC due to mutations in PTPRH.
PLoS genetics
Swiatnicki MR,Rennhack JP,Ortiz MMO,Hollern DP,Perry AV,Kubiak R,Riveria Riveria SM,O'Reilly S,Andrechek ER
Thu Sep 01 00:00:00 UTC 2022
Hyperhomocysteinemia Promotes Cardiac Hypertrophy in Hypertension.
Oxidative medicine and cellular longevity
Deng Y,Li Z,An X,Fan R,Wang Y,Li J,Yang X,Liao J,Xia Y
Wed Sep 07 00:00:00 UTC 2022
Silencing lncRNA 93358 Inhibits the Apoptosis of Myocardial Cells in Myocardial Infarction Rats by Inducing the Expression of SLC8A1.
BioMed research international
Cai J,Wang X,Liao W,Zhong Y,Chen L,Zhang Z
Tue Jul 19 00:00:00 UTC 2022
Quantitative proteomics and functional analysis identified novel targets for missed abortion.
Experimental cell research
Chen X,Zheng Q,Ji L,Zhuang Y,Yu H,Cheng X,Han Y,Lv J,Zheng B,Zheng Y,Yu J
Mon Aug 15 00:00:00 UTC 2022
Sesamin Activates Skeletal Muscle FNDC5 Expression and Increases Irisin Secretion via the SIRT1 Signaling Pathway.
Journal of agricultural and food chemistry
Kou G,Li P,Shi Y,Traore SS,Shi X,Amoah AN,Cui Z,Lyu Q
Wed Jun 29 00:00:00 UTC 2022
Attachment of -tBu groups to aza-BODIPY core at 3,5-sites with ultra-large Stokes shift to enhance photothermal therapy through apoptosis mechanism.
Materials today. Bio
Li R,Ren J,Zhang D,Lv M,Wang Z,Wang H,Zhang S,Du J,Jiang XD,Wang G
Thu Dec 01 00:00:00 UTC 2022
Sleeve Gastrectomy-Induced AMPK Activation Attenuates Diabetic Cardiomyopathy by Maintaining Mitochondrial Homeostasis via NR4A1 Suppression in Rats.
Frontiers in physiology
Li S,Dong S,Xu Q,Shi B,Li L,Zhang W,Zhu J,Cheng Y,Zhang G,Zhong M
Sat Apr 02 00:00:00 UTC 2022
BCAA-BCKA axis regulates WAT browning through acetylation of PRDM16.
Nature metabolism
Ma QX,Zhu WY,Lu XC,Jiang D,Xu F,Li JT,Zhang L,Wu YL,Chen ZJ,Yin M,Huang HY,Lei QY
Sat Jan 01 00:00:00 UTC 2022
The preventive and therapeutic effect of repetitive transcranial magnetic stimulation on radiation-induced brain injury in mice.
International journal of radiation biology
Liu LY,Qin TZ,Guo L,Rong-Rong H,Jing YT,Lai PP,Xue YZ,Ding GR
Wed Jul 27 00:00:00 UTC 2022
Chronic exposure of alcohol triggers microglia-mediated synaptic elimination inducing cognitive impairment.
Experimental neurology
Lan L,Wang H,Zhang X,Shen Q,Li X,He L,Rong X,Peng J,Mo J,Peng Y
Fri Jul 01 00:00:00 UTC 2022
The mechanism on Prevotella melaninogenica promoting the inflammatory progression of oral lichen planus.
Clinical and experimental immunology
Xu P,Shao RR,Zhang S,Tan ZW,Guo YT,He Y
Fri Aug 19 00:00:00 UTC 2022
The protective effects of Xuebijing injection on intestinal injuries of mice exposed to irradiation.
Animal models and experimental medicine
Dong Y,Zhang Y,Wang X,Li W,Zhang J,Lu L,Dong H,Fan S,Meng A,Li D
Thu Dec 01 00:00:00 UTC 2022
An efficient strategy for digging protein-protein interactions for rational drug design - A case study with HIF-1α/VHL.
European journal of medicinal chemistry
Xue X,Kang JB,Yang X,Li N,Chang L,Ji J,Meng XK,Zhang HQ,Zhong Y,Yu SP,Wu WY,Wang XL,Li NG,Sun SL
Wed Jan 05 00:00:00 UTC 2022
Type IIB PKA is highly expressed in β cells and controls cell proliferation via regulating Cyclin D1 expression.
The FEBS journal
Jiang Y,Zhu L,Wu D,Ni Y,Huang C,Ye H,Yang Y,Liu R,Li Y
Sun May 01 00:00:00 UTC 2022
Iron metabolism protein transferrin receptor 1 involves in cervical cancer progression by affecting gene expression and alternative splicing in HeLa cells.
Genes & genomics
Huang N,Wei Y,Cheng Y,Wang X,Wang Q,Chen D,Li W
Wed Jun 01 00:00:00 UTC 2022
Qingwei San treats oral ulcer subjected to stomach heat syndrome in db/db mice by targeting TLR4/MyD88/NF-κB pathway.
Chinese medicine
Shi L,An Y,Cheng L,Li Y,Li H,Wang C,Lv Y,Duan Y,Dai H,He C,Zhang H,Huang Y,Fu W,Wang S,Zhao B,Wang Y,Zhao Y
Tue Jan 04 00:00:00 UTC 2022
Cellular communication network factor 1 interlinks autophagy and ERK signaling to promote osteogenesis of periodontal ligament stem cells.
Journal of periodontal research
Li Z,Wu Z,Xi X,Zhao F,Liu H,Liu D
Thu Dec 01 00:00:00 UTC 2022
Lysyl oxidase-like 2 inhibitor rescues D-galactose-induced skeletal muscle fibrosis.
Aging cell
Wu Y,Wu Y,Yang Y,Yu J,Wu J,Liao Z,Guo A,Sun Y,Zhao Y,Chen J,Xiao Q
Fri Jul 01 00:00:00 UTC 2022
Vagus nerve stimulation alleviated cerebral ischemia and reperfusion injury in rats by inhibiting pyroptosis via α7 nicotinic acetylcholine receptor.
Cell death discovery
Tang H,Li J,Zhou Q,Li S,Xie C,Niu L,Ma J,Li C
Tue Feb 08 00:00:00 UTC 2022
FAEE exerts a protective effect against osteoporosis by regulating the MAPK signalling pathway.
Pharmaceutical biology
Wang MY,An MF,Fan MS,Zhang SS,Sun ZR,Zhao YL,Xiang ZM,Sheng J
Thu Dec 01 00:00:00 UTC 2022
JNK and Jag1/Notch2 co-regulate CXCL16 to facilitate cypermethrin-induced kidney damage.
Ecotoxicology and environmental safety
Liu C,Wu M,Qu J,Huang X,Zeng Q,Ha M
Wed Jun 15 00:00:00 UTC 2022
The role of E3 ubiquitin ligase WWP2 and the regulation of PARP1 by ubiquitinated degradation in acute lymphoblastic leukemia.
Cell death discovery
Lu X,Huang X,Xu H,Lu S,You S,Xu J,Zhan Q,Dong C,Zhang N,Zhang Y,Cao L,Zhang X,Zhang N,Zhang L
Tue Oct 18 00:00:00 UTC 2022
Role of PARP-1 in Human Cytomegalovirus Infection and Functional Partners Encoded by This Virus.
Viruses
Zhang W,Guo J,Chen Q
Thu Sep 15 00:00:00 UTC 2022
Selective Inhibition of Histone Deacetylase Class IIa With MC1568 Ameliorates Podocyte Injury.
Frontiers in medicine
He X,Sun T,Zhang P,Xia Z,Gao C,Ren H,Ji D
Sat Jul 16 00:00:00 UTC 2022
Hydrogen improves exercise endurance in rats by promoting mitochondrial biogenesis.
Genomics
Luo M,Lu J,Li C,Wen B,Chu W,Dang X,Zhang Y,An G,Wang J,Fan R,Chen X
Tue Nov 01 00:00:00 UTC 2022
Map of Enteropathogenic Escherichia coli Targets Mitochondria and Triggers DRP-1-Mediated Mitochondrial Fission and Cell Apoptosis in Bovine Mastitis.
International journal of molecular sciences
Li Y,Zhu Y,Chu B,Liu N,Chen S,Wang J,Zou Y
Thu Apr 28 00:00:00 UTC 2022
Heart-targeted amelioration of sepsis-induced myocardial dysfunction by microenvironment responsive nitric oxide nanogenerators in situ.
Journal of nanobiotechnology
Ouyang M,Ouyang X,Peng Z,Liu M,Xu G,Zou Z,Zhang M,Shang Q
Tue Jun 07 00:00:00 UTC 2022
TRPM8 contributes to sex dimorphism by promoting recovery of normal sensitivity in a mouse model of chronic migraine.
Nature communications
Alarcón-Alarcón D,Cabañero D,de Andrés-López J,Nikolaeva-Koleva M,Giorgi S,Fernández-Ballester G,Fernández-Carvajal A,Ferrer-Montiel A
Sat Oct 22 00:00:00 UTC 2022
Increased m6A-RNA methylation and FTO suppression is associated with myocardial inflammation and dysfunction during endotoxemia in mice.
Molecular and cellular biochemistry
Dubey PK,Patil M,Singh S,Dubey S,Ahuja P,Verma SK,Krishnamurthy P
Sat Jan 01 00:00:00 UTC 2022
Golgi Phosphoprotein 3 Confers Radioresistance via Stabilizing EGFR in Lung Adenocarcinoma.
International journal of radiation oncology, biology, physics
Chen G,Kong P,Yang M,Hu W,Prise KM,Yu KN,Cui S,Qin F,Meng G,Almahi WA,Nie L,Han W
Fri Apr 01 00:00:00 UTC 2022
Trichostatin D as a Novel KLF2 Activator Attenuates TNFα-Induced Endothelial Inflammation.
International journal of molecular sciences
Lei L,Chen M,Wang C,Jiang X,Li Y,Wang W,Li S,Zhao L,Sheng R,Han J,Zhang Y,Chen Y,Yan B,Wu Y,Yu L,Si S,Xu Y
Thu Nov 03 00:00:00 UTC 2022
Inhibitory effect of tanshinone IIA, resveratrol and silibinin on enterovirus 68 production through inhibiting ATM and DNA-PK pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Su Y,Wu T,Yu XY,Huo WB,Wang SH,Huan C,Liu YM,Liu JM,Cui MN,Li XH,Yu JH
Sun May 01 00:00:00 UTC 2022
Effects of ALA-PDT on the macrophages in wound healing and its related mechanisms in vivo and in vitro.
Photodiagnosis and photodynamic therapy
Li L,Yang Y,Yang Z,Zheng M,Luo G,He W,Yin R
Wed Jun 01 00:00:00 UTC 2022
Angiotensin-converting enzyme 2 identifies immuno-hot tumors suggesting angiotensin-(1-7) as a sensitizer for chemotherapy and immunotherapy in breast cancer.
Biological procedures online
Mei J,Cai Y,Xu R,Yu X,Han X,Weng M,Chen L,Ma T,Gao T,Gao F,Xia T,Zhu Y,Zhang Y
Tue Oct 25 00:00:00 UTC 2022
Chemical profile and miscarriage prevention evaluation of Jiao-Ai Decoction, a classical traditional Chinese formula.
Journal of pharmaceutical and biomedical analysis
Zhang M,Geng T,Jing SH,Li L,Wang K,Zhang Q,Zhang Y,Zhou SK,Gao P,Ding AW,Zhang L
Fri Aug 05 00:00:00 UTC 2022
Label-free LC-MS/MS proteomics analyses reveal CLIC1 as a predictive biomarker for bladder cancer staging and prognosis.
Frontiers in oncology
Wang W,Huang G,Lin H,Ren L,Fu L,Mao X
Wed Jan 24 00:00:00 UTC 2024
Sodium hydrosulfide reverses β2-microglobulin-induced depressive-like behaviors of male Sprague-Dawley rats: Involving improvement of synaptic plasticity and enhancement of Warburg effect in hippocampus.
Behavioural brain research
Yang SQ,Tang YY,Zeng D,Tian Q,Wei HJ,Wang CY,Zhang P,Chen YJ,Zou W,Tang XQ
Mon Jan 24 00:00:00 UTC 2022
Punicalagin Alleviates Psoriasis by Inhibiting NF-κB-Mediated IL-1β Transcription and Caspase-1-Regulated IL-1β Secretion.
Frontiers in pharmacology
Tang L,Li T,Zhang B,Zhang Z,Sun X,Zhu Y,Feng B,Su Z,Yang L,Li H,Liu H,Chen Y,Dai Z,Zheng X,Li M,Li C,Zhao J,Qiu X,Ye S,Liu H,Zheng G,Li B,Lu C
Wed Feb 16 00:00:00 UTC 2022
Berberine prevents lethal EV71 neurological infection in newborn mice.
Frontiers in pharmacology
Cui G,Wang H,Yang C,Zhou X,Wang J,Wang T,Ma T
Fri Nov 18 00:00:00 UTC 2022
The PROTAC selectively degrading Bcl-xL represents a novel Hedgehog pathway inhibitor with capacity of combating resistance to Smoothened inhibitors while sparing bone growth.
Theranostics
Zhang S,Chen Y,Xu Z,Yang J,Sun R,Wang J,Sun Y,Jiang B,Yang X,Tan W
Tue Nov 29 00:00:00 UTC 2022
Cypermethrin inhibits proliferation of Sertoli cells through AR involving DAB2IP/PI3K/AKT signaling pathway in vitro.
Toxicology research
Wang Q,Wang XX,Xie JF,Yao TT,Xu LL,Wang LS,Yu Y,Xu LC
Mon Aug 01 00:00:00 UTC 2022
Hypoxia facilitates proliferation of smooth muscle cells derived from pluripotent stem cells for vascular tissue engineering.
Journal of tissue engineering and regenerative medicine
Fang L,Mei J,Yao B,Liu J,Liu P,Wang X,Zhou J,Lin Z
Mon Aug 01 00:00:00 UTC 2022
Metabolic Syndrome Ameliorated by 4-Methylesculetin by Reducing Hepatic Lipid Accumulation.
International journal of molecular sciences
Li L,Zhu G,Fu G,Zha W,Li H
Fri Sep 09 00:00:00 UTC 2022
Aberrant translation regulated by METTL1/WDR4-mediated tRNA N7-methylguanosine modification drives head and neck squamous cell carcinoma progression.
Cancer communications (London, England)
Chen J,Li K,Chen J,Wang X,Ling R,Cheng M,Chen Z,Chen F,He Q,Li S,Zhang C,Jiang Y,Chen Q,Wang A,Chen D
Tue Mar 01 00:00:00 UTC 2022
Ejection of damaged mitochondria and their removal by macrophages ensure efficient thermogenesis in brown adipose tissue.
Cell metabolism
Rosina M,Ceci V,Turchi R,Chuan L,Borcherding N,Sciarretta F,Sánchez-Díaz M,Tortolici F,Karlinsey K,Chiurchiù V,Fuoco C,Giwa R,Field RL,Audano M,Arena S,Palma A,Riccio F,Shamsi F,Renzone G,Verri M,Crescenzi A,Rizza S,Faienza F,Filomeni G,Kooijman S,Rufini S,de Vries AAF,Scaloni A,Mitro N,Tseng YH,Hidalgo A,Zhou B,Brestoff JR,Aquilano K,Lettieri-Barbato D
Tue Apr 05 00:00:00 UTC 2022
cGAS-STING mediates cytoplasmic mitochondrial-DNA-induced inflammatory signal transduction during accelerated senescence of pancreatic β-cells induced by metabolic stress.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hu H,Zhao R,He Q,Cui C,Song J,Guo X,Zang N,Yang M,Zou Y,Yang J,Li J,Wang L,Xia L,Wang L,He F,Hou X,Yan F,Chen L
Sun May 01 00:00:00 UTC 2022
NAP1L5 targeting combined with MYH9 Inhibit HCC progression through PI3K/AKT/mTOR signaling pathway.
Aging
Zhao R,Ge Y,Gong Y,Li B,Xiao B,Zuo S
Fri Nov 11 00:00:00 UTC 2022
ID2 inhibits innate antiviral immunity by blocking TBK1- and IKKε-induced activation of IRF3.
Science signaling
Yu C,Wang B,Zhu Y,Zhang C,Ren L,Lei X,Xiang Z,Zhou Z,Huang H,Wang J,Zhao Z
Tue Jan 04 00:00:00 UTC 2022
Phosphorylation of Ser82 on IRF3 acts as negative-feedback regulation of IRF3-dependent innate immunity.
The international journal of biochemistry & cell biology
Cheng Q,Yuan L,Guo J,Guo D,Liu X,Li S
Thu Sep 01 00:00:00 UTC 2022
SMAD4 Inhibits Granulosa Cell Apoptosis via the miR-183-96-182 Cluster and FoxO1 Axis.
Reproductive sciences (Thousand Oaks, Calif.)
Yao W,Wang S,Du X,Lin C,Zhang J,Pan Z,Li Q
Sun May 01 00:00:00 UTC 2022
Dexmedetomidine attenuates perioperative neurocognitive disorders by suppressing hippocampal neuroinflammation and HMGB1/RAGE/NF-κB signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang J,Xin Y,Chu T,Liu C,Xu A
Wed Jun 01 00:00:00 UTC 2022
Cuproptosis status affects treatment options about immunotherapy and targeted therapy for patients with kidney renal clear cell carcinoma.
Frontiers in immunology
Zhang G,Chen X,Fang J,Tai P,Chen A,Cao K
Wed Feb 15 00:00:00 UTC 2023
Toddalolactone protects against osteoarthritis by ameliorating chondrocyte inflammation and suppressing osteoclastogenesis.
Chinese medicine
Xu Y,Xue S,Zhang T,Jin X,Wang C,Lu H,Zhong Y,Chen H,Zhu L,Ma J,Sang W
Sat Feb 05 00:00:00 UTC 2022
Copy Number Analyses Identified a Novel Gene: APOBEC3A Related to Lipid Metabolism in the Pathogenesis of Preeclampsia.
Frontiers in cardiovascular medicine
Liu N,Guo YN,Wang XJ,Ma J,He YT,Zhang F,He H,Xie JL,Zhuang X,Liu M,Sun JH,Chen Y,Lin JH,Gong LK,Wang BS
Sat Jul 16 00:00:00 UTC 2022
Cdk5 Promotes Mitochondrial Fission via Drp1 Phosphorylation at S616 in Chronic Ethanol Exposure-Induced Cognitive Impairment.
Molecular neurobiology
Liu D,Li J,Rong X,Li J,Peng Y,Shen Q
Thu Dec 01 00:00:00 UTC 2022
Extracellular CIRP Upregulates Proinflammatory Cytokine Expression via the NF-kappaB and ERK1/2 Signaling Pathways in Psoriatic Keratinocytes.
Mediators of inflammation
Zhang X,Wang S,Wang W,Song L,Feng S,Wang J,Kang T,Yang P,Wang N,Yang P,Bai R,Shao Y,Zheng Y
Mon Sep 19 00:00:00 UTC 2022
Synthetic Retinoid Kills Drug-Resistant Cancer Stem Cells via Inducing RARγ-Translocation-Mediated Tension Reduction and Chromatin Decondensation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Dong Q,An Q,Zhang C,Mohagheghian E,Niu B,Qi F,Wei F,Chen S,Chen X,Wang A,Cao X,Wang N,Chen J
Tue Nov 01 00:00:00 UTC 2022
Regulation of cohesin-mediated chromosome folding by PDS5 in mammals.
EMBO reports
Yu D,Chen G,Wang Y,Wang Y,Lin R,Liu N,Zhu P,Liu H,Hu T,Feng R,Feng H,Lan F,Cai J,Chen H
Mon Nov 07 00:00:00 UTC 2022
Dysregulation of SIRT3 SUMOylation Confers AML Chemoresistance via Controlling HES1-Dependent Fatty Acid Oxidation.
International journal of molecular sciences
Zhang Y,Shen Y,Wei W,Wang W,Jiang D,Ren Y,Peng Z,Fan Q,Cheng J,Ma J
Wed Jul 27 00:00:00 UTC 2022
Dual effects of bisphenol A on wound healing, involvement of estrogen receptor β.
Ecotoxicology and environmental safety
Wen X,Zhu M,Li Z,Li T,Xu X
Tue Feb 01 00:00:00 UTC 2022
Epoxyeicosatrienoic Acids Inhibit the Activation of Murine Fibroblasts by Blocking the TGF-β1-Smad2/3 Signaling in a PPARγ-Dependent Manner.
Oxidative medicine and cellular longevity
Tao JH,Liu T,Zhang CY,Zu C,Yang HH,Liu YB,Yang JT,Zhou Y,Guan CX
Thu Oct 27 00:00:00 UTC 2022
DsbA-L interacts with VDAC1 in mitochondrion-mediated tubular cell apoptosis and contributes to the progression of acute kidney disease.
EBioMedicine
Li X,Pan J,Li H,Li G,Liu B,Tang X,Liu X,He Z,Peng Z,Zhang H,Wang L,Li Y,Xiang X,Chai X,Yuan Y,Zheng P,Zhang D
Tue Feb 01 00:00:00 UTC 2022
Identifying Potential Tumor Antigens and Antigens-Related Subtypes in Hepatocellular Carcinoma for mRNA Vaccine Development.
Journal of oncology
Liao W,Shi Z,Tang H,Wu T,Zhang C,He Y,Zou R,Wang L
Sat Sep 10 00:00:00 UTC 2022
CHMFL-BMX-078, a BMX inhibitor, overcomes the resistance of melanoma to vemurafenib via inhibiting AKT pathway.
Chemico-biological interactions
Jiang S,Jiang T,Huang H,Chen X,Li L,Wang Z,Fei J,Liu C,Liu Z,Cheng Y
Wed Jan 05 00:00:00 UTC 2022
Kaempferol prevents angiogenesis of rat intestinal microvascular endothelial cells induced by LPS and TNF-α via inhibiting VEGF/Akt/p38 signaling pathways and maintaining gut-vascular barrier integrity.
Chemico-biological interactions
Yu R,Zhong J,Zhou Q,Ren W,Liu Z,Bian Y
Sat Oct 01 00:00:00 UTC 2022
The expression of m6A enzymes in the hippocampus of diabetic cognitive impairment mice and the possible improvement of YTHDF1.
Brain research
Li M,Zhong X,Zhao Z,Zeng Z,Yuan Q,Xiao X,Zu X,Liu J
Tue Feb 15 00:00:00 UTC 2022
Ubiquitin-like modifier 1 ligating enzyme 1 relieves cisplatin-induced premature ovarian failure by reducing endoplasmic reticulum stress in granulosa cells.
Reproductive biology and endocrinology : RB&E
Tang X,Dong H,Fang Z,Li J,Yang Q,Yao T,Pan Z
Tue May 24 00:00:00 UTC 2022
Therapeutic effects of shaogan fuzi decoction in rheumatoid arthritis: Network pharmacology and experimental validation.
Frontiers in pharmacology
Shi L,Zhao Y,Feng C,Miao F,Dong L,Wang T,Stalin A,Zhang J,Tu J,Liu K,Sun W,Wu J
Wed Sep 07 00:00:00 UTC 2022
ANXA1: An Important Independent Prognostic Factor and Molecular Target in Glioma.
Frontiers in genetics
Zhang D,Wang W,Zhou H,Su L,Han X,Zhang X,Han W,Wang Y,Xue X
Sat Jul 16 00:00:00 UTC 2022
Deficiency of proline/serine-rich coiled-coil protein 1 (PSRC1) accelerates trimethylamine N-oxide-induced atherosclerosis in ApoE-/- mice.
Journal of molecular and cellular cardiology
Luo T,Liu D,Guo Z,Chen P,Guo Z,Ou C,Chen M
Thu Sep 01 00:00:00 UTC 2022
Krill Oil Inhibits NLRP3 Inflammasome Activation in the Prevention of the Pathological Injuries of Diabetic Cardiomyopathy.
Nutrients
Sun X,Sun X,Meng H,Wu J,Guo X,Du L,Wu H
Sat Jan 15 00:00:00 UTC 2022
MxB inhibits long interspersed element type 1 retrotransposition.
PLoS genetics
Huang Y,Xu F,Mei S,Liu X,Zhao F,Wei L,Fan Z,Hu Y,Wang L,Ai B,Cen S,Liang C,Guo F
Tue Feb 01 00:00:00 UTC 2022
Melatonin protects oocytes from cadmium exposure-induced meiosis defects by changing epigenetic modification and enhancing mitochondrial morphology in the mouse.
Ecotoxicology and environmental safety
Qu J,Wang Q,Niu H,Sun X,Ji D,Li Y
Thu Dec 15 00:00:00 UTC 2022
Quantitative proteomics analysis to assess protein expression levels in the ovaries of pubescent goats.
BMC genomics
Qin P,Ye J,Gong X,Yan X,Lin M,Lin T,Liu T,Li H,Wang X,Zhu Y,Li X,Liu Y,Li Y,Ling Y,Zhang X,Fang F
Wed Jul 13 00:00:00 UTC 2022
Proteomic and Biochemical Approaches Elucidate the Role of Millimeter-Wave Irradiation in Wheat Growth under Flooding Stress.
International journal of molecular sciences
Komatsu S,Tsutsui Y,Furuya T,Yamaguchi H,Hitachi K,Tsuchida K,Tani M
Thu Sep 08 00:00:00 UTC 2022
Apextrin from Ruditapes philippinarum functions as pattern recognition receptor and modulates NF-κB pathway.
International journal of biological macromolecules
Jiang K,Nie H,Yin Z,Yan X,Li Q
Mon Aug 01 00:00:00 UTC 2022
Identification of super-enhancer-driven peptidyl arginine deiminases as potential biomarkers and therapeutic targets for osimertinib-resistant non-small cell lung cancer.
Frontiers in pharmacology
Li H,Muhetaer G,Xie Y,Yao K,Ma Q,Guan H,Xing S,Huang X,Zhou J
Sat Dec 10 00:00:00 UTC 2022
Prediction of Prognosis and Recurrence of Bladder Cancer by ECM-Related Genes.
Journal of immunology research
Zhao H,Chen Z,Fang Y,Su M,Xu Y,Wang Z,Gyamfi MA,Zhao J
Mon Apr 25 00:00:00 UTC 2022
Discovery of Novel Phosphoinositide-3-Kinase α Inhibitors with High Selectivity, Excellent Bioavailability, and Long-Acting Efficacy for Gastric Cancer.
Journal of medicinal chemistry
Hou Y,Zhang F,Min W,Yuan K,Kuang W,Wang X,Zhu Y,Sun C,Xia F,Wang Y,Zhang H,Wang L,Yang P
Thu Jul 28 00:00:00 UTC 2022
Resveratrol attenuates inflammation and apoptosis through alleviating endoplasmic reticulum stress via Akt/mTOR pathway in fungus-induced allergic airways inflammation.
International immunopharmacology
Wang S,Gong L,Mo Y,Zhang J,Jiang Z,Tian Z,Shao C
Tue Feb 01 00:00:00 UTC 2022
A novel HCC prognosis predictor PDSS1 affects the cell cycle through the STAT3 signaling pathway in HCC.
Frontiers in oncology
Rao Z,Li H,Yao W,Wang Q,Ma B,Xue D,Meng X
Tue Aug 16 00:00:00 UTC 2022
S1PR3, as a Core Protein Related to Ischemic Stroke, is Involved in the Regulation of Blood-Brain Barrier Damage.
Frontiers in pharmacology
Fan X,Chen H,Xu C,Wang Y,Yin P,Li M,Tang Z,Jiang F,Wei W,Song J,Li G,Zhong D
Sat Jul 16 00:00:00 UTC 2022
Optical Visualization of Red-GQDs' Organelles Distribution and Localization in Living Cells.
Frontiers in pharmacology
Hu H,Li P,Qiu J,Zhao M,Kuang M,Zhang Z,Wang D
Tue Aug 02 00:00:00 UTC 2022
Differential requirements of IQUB for the assembly of radial spoke 1 and the motility of mouse cilia and flagella.
Cell reports
Zhang X,Xiao Z,Zhang J,Xu C,Liu S,Cheng L,Zhou S,Zhao S,Zhang Y,Wu J,Wang Y,Liu M
Tue Nov 22 00:00:00 UTC 2022
Caspase-1-mediated extracellular vesicles derived from pyroptotic alveolar macrophages promote inflammation in acute lung injury.
International journal of biological sciences
Qin X,Zhou Y,Jia C,Chao Z,Qin H,Liang J,Liu X,Liu Z,Sun T,Yuan Y,Zhang H
Fri Apr 15 00:00:00 UTC 2022
Difenoconazole causes spleen tissue damage and immune dysfunction of carp through oxidative stress and apoptosis.
Ecotoxicology and environmental safety
Liu F,Li X,Bello BK,Zhang T,Yang H,Wang K,Dong J
Wed Jun 01 00:00:00 UTC 2022
Dexpramipexole Attenuates White Matter Injury to Facilitate Locomotion and Motor Coordination Recovery via Reducing Ferroptosis after Intracerebral Hemorrhage.
Oxidative medicine and cellular longevity
Wang B,Zhang X,Zhong J,Wang S,Zhang C,Li M,Hu Q,Wang S,Chen L,Chen W,Ge H,Feng H
Tue Aug 16 00:00:00 UTC 2022
Defective autophagy triggered by arterial cyclic stretch promotes neointimal hyperplasia in vein grafts via the p62/nrf2/slc7a11 signaling pathway.
Journal of molecular and cellular cardiology
Chen Y,Bao M,Liu JT,Bao H,Zhang SM,Lou Y,Qi YX
Thu Dec 01 00:00:00 UTC 2022
Activating PPARβ/δ Protects against Endoplasmic Reticulum Stress-Induced Astrocytic Apoptosis via UCP2-Dependent Mitophagy in Depressive Model.
International journal of molecular sciences
Ji J,Li S,Jiang Z,Yu J,Sun Y,Cai Z,Dong Y,Sun X
Fri Sep 16 00:00:00 UTC 2022
The plant hormone abscisic acid stimulates megakaryocyte differentiation from human iPSCs in vitro.
Platelets
Huang W,Gu H,Zhan Z,Wang R,Song L,Zhang Y,Zhang Y,Li S,Li J,Zang Y,Li Y,Qian B
Sun Apr 03 00:00:00 UTC 2022
Chicken ISG12(2) attenuates Newcastle disease virus and enhances the efficiency of Newcastle disease vaccine via activating immune pathways.
Transboundary and emerging diseases
Liu H,Tian J,Lu K,Li Y,Guan Z,Cao X,Li X,Chang Z,Wang X,Sa X,Yang Z
Thu Sep 01 00:00:00 UTC 2022
Proteomic characterization of gastric cancer response to chemotherapy and targeted therapy reveals new therapeutic strategies.
Nature communications
Li Y,Xu C,Wang B,Xu F,Ma F,Qu Y,Jiang D,Li K,Feng J,Tian S,Wu X,Wang Y,Liu Y,Qin Z,Liu Y,Qin J,Song Q,Zhang X,Sujie A,Huang J,Liu T,Shen K,Zhao JY,Hou Y,Ding C
Thu Sep 29 00:00:00 UTC 2022
Nav1.8 in keratinocytes contributes to ROS-mediated inflammation in inflammatory skin diseases.
Redox biology
Zhang Y,Li Y,Zhou L,Yuan X,Wang Y,Deng Q,Deng Z,Xu S,Wang Q,Xie H,Li J
Thu Sep 01 00:00:00 UTC 2022
MiR-135a-5p suppresses trophoblast proliferative, migratory, invasive, and angiogenic activity in the context of unexplained spontaneous abortion.
Reproductive biology and endocrinology : RB&E
Lu Y,Zhang X,Li X,Deng L,Wei C,Yang D,Tan X,Pan W,Pang L
Tue May 24 00:00:00 UTC 2022
Hyperglycemia promotes myocardial dysfunction via the ERS-MAPK10 signaling pathway in db/db mice.
Laboratory investigation; a journal of technical methods and pathology
Deng YW,Liu F,Li ZT,Gao JH,Zhao Y,Yang XL,Xia YL
Tue Nov 01 00:00:00 UTC 2022
O-GlcNAc transferase promotes the nuclear localization of the focal adhesion-associated protein Zyxin to regulate UV-induced cell death.
The Journal of biological chemistry
Zhao Y,Yue S,Zhou X,Guo J,Ma S,Chen Q
Fri Apr 01 00:00:00 UTC 2022
Proteomic-Based Approach Reveals the Involvement of Apolipoprotein A-I in Related Phenotypes of Autism Spectrum Disorder in the BTBR Mouse Model.
International journal of molecular sciences
Li Q,Shi Y,Li X,Yang Y,Zhang X,Xu L,Ma Z,Wang J,Fan L,Wu L
Sun Dec 04 00:00:00 UTC 2022
The bibenzyl derivatives of Dendrobium officinale prevent UV-B irradiation induced photoaging via SIRT3.
Natural products and bioprospecting
Chen DK,Shao HY,Yang L,Hu JM
Thu Jan 27 00:00:00 UTC 2022
LncRNA ALKBH3-AS1 enhances ALKBH3 mRNA stability to promote hepatocellular carcinoma cell proliferation and invasion.
Journal of cellular and molecular medicine
Lu Q,Wang H,Lei X,Ma Q,Zhao J,Sun W,Guo C,Huang D,Xu Q
Sat Oct 01 00:00:00 UTC 2022
RNA-m6A modification of HDGF mediated by Mettl3 aggravates the progression of atherosclerosis by regulating macrophages polarization via energy metabolism reprogramming.
Biochemical and biophysical research communications
Zheng L,Chen X,Yin Q,Gu J,Chen J,Chen M,Zhang Y,Dong M,Jiang H,Yin N,Chen H,Li X
Tue Dec 20 00:00:00 UTC 2022
mmu-lncRNA 121686/hsa-lncRNA 520657 induced by METTL3 drive the progression of AKI by targeting miR-328-5p/HtrA3 signaling axis.
Molecular therapy : the journal of the American Society of Gene Therapy
Pan J,Xie Y,Li H,Li X,Chen J,Liu X,Zhou J,Tang X,He Z,Peng Z,Zhang H,Li Y,Xiang X,Yuan Y,Zhang D
Wed Dec 07 00:00:00 UTC 2022
The lncRNA MIAT regulates CPT-1a mediated cardiac hypertrophy through m6A RNA methylation reading protein Ythdf2.
Cell death discovery
Yang Y,Mbikyo MB,Zhang J,Zhang Y,Zhang N,Li Z
Tue Apr 05 00:00:00 UTC 2022
ORP5 promotes tumor metastasis via stabilizing c-Met in renal cell carcinoma.
Cell death discovery
Song L,Zhang L,Zhou Y,Shao X,Xu Y,Pei D,Wang Q
Thu Apr 21 00:00:00 UTC 2022
Cortistatin attenuates titanium particle-induced osteolysis through regulation of TNFR1-ROS-caspase-3 signaling in osteoblasts.
Annals of the New York Academy of Sciences
Cao J,Ma X,Liu L,Zhang G,Wu Y,Fu Y,Gong A,Yang Z,Zhao Y,Zhang L,Li Y
Fri Jul 01 00:00:00 UTC 2022
The neuroprotective effects of GPR55 against hippocampal neuroinflammation and impaired adult neurogenesis in CSDS mice.
Neurobiology of disease
Shen SY,Yu R,Li W,Liang LF,Han QQ,Huang HJ,Li B,Xu SF,Wu GC,Zhang YQ,Yu J
Fri Jul 01 00:00:00 UTC 2022
Overexpression of PYGO1 promotes early cardiac lineage development in human umbilical cord mesenchymal stromal/stem cells by activating the Wnt/β-catenin pathway.
Human cell
Shen J,Wu X,Zhu P,Zhuang J,Qin B,Sun F,Yuan W,Fan X,Jiang Z,Li F,Li Y,Wang Y,Zhao M
Tue Nov 01 00:00:00 UTC 2022
Metformin Protects against Spinal Cord Injury and Cell Pyroptosis via AMPK/NLRP3 Inflammasome Pathway.
Analytical cellular pathology (Amsterdam)
Yuan Y,Fan X,Guo Z,Zhou Z,Gao W
Fri Apr 08 00:00:00 UTC 2022
Chronic exposure to biomass ambient particulate matter triggers alveolar macrophage polarization and activation in the rat lung.
Journal of cellular and molecular medicine
Wang S,Chen Y,Hong W,Li B,Zhou Y,Ran P
Tue Feb 01 00:00:00 UTC 2022
Effect of miR-143-3p from Extracellular Vesicles of Porcine Uterine Luminal Fluid on Porcine Trophoblast Cells.
Animals : an open access journal from MDPI
Ding Y,Hu Q,Gan J,Zang X,Gu T,Wu Z,Cai G,Hong L
Fri Dec 02 00:00:00 UTC 2022
Interleukin 33-mediated inhibition of A-type K+ channels induces sensory neuronal hyperexcitability and nociceptive behaviors in mice.
Theranostics
Wang Y,Wang X,Qi R,Lu Y,Tao Y,Jiang D,Sun Y,Jiang X,Liu C,Zhang Y,Tao J
Thu Apr 21 00:00:00 UTC 2022
The Hair Growth-Promoting Effect of Gardenia florida Fruit Extract and Its Molecular Regulation.
Evidence-based complementary and alternative medicine : eCAM
Liu X,Ji T,Hu H,Lv X,Zhu G
Thu Oct 06 00:00:00 UTC 2022
Deacetylation of Caveolin-1 by Sirt6 induces autophagy and retards high glucose-stimulated LDL transcytosis and atherosclerosis formation.
Metabolism: clinical and experimental
Zhao Y,Jia X,Yang X,Bai X,Lu Y,Zhu L,Cheng W,Shu M,Zhu Y,Du X,Wang L,Shu Y,Song Y,Jin S
Wed Jun 01 00:00:00 UTC 2022
Sclerostin Suppression Facilitates Uveal Melanoma Progression Through Activating Wnt/β-Catenin Signaling Via Binding to Membrane Receptors LRP5/LRP6.
Frontiers in oncology
Wang H,Zhao S,Liu Y,Sun F,Huang X,Wu T
Sat Jul 16 00:00:00 UTC 2022
Sclerostin Suppression Facilitates Uveal Melanoma Progression Through Activating Wnt/β-Catenin Signaling Via Binding to Membrane Receptors LRP5/LRP6.
Frontiers in oncology
Wang H,Zhao S,Liu Y,Sun F,Huang X,Wu T
Sat Jul 16 00:00:00 UTC 2022
circEXOC6B interacting with RRAGB, an mTORC1 activator, inhibits the progression of colorectal cancer by antagonizing the HIF1A-RRAGB-mTORC1 positive feedback loop.
Molecular cancer
Li X,Wang J,Lin W,Yuan Q,Lu Y,Wang H,Chen Y,Chen L,Dai P,Long H,Li X
Thu Jun 23 00:00:00 UTC 2022
HIF-1/2α-Activated RNF146 Enhances the Proliferation and Glycolysis of Hepatocellular Carcinoma Cells via the PTEN/AKT/mTOR Pathway.
Frontiers in cell and developmental biology
Shen G,Wang H,Zhu N,Lu Q,Liu J,Xu Q,Huang D
Sat Jul 16 00:00:00 UTC 2022
The mechanism of dioscin preventing lung cancer based on network pharmacology and experimental validation.
Journal of ethnopharmacology
Xi P,Niu Y,Zhang Y,Li W,Gao F,Gu W,Kui F,Liu Z,Lu L,Du G
Tue Jun 28 00:00:00 UTC 2022
Local Application of Krill Oil Accelerates the Healing of Artificially Created Wounds in Diabetic Mice.
Nutrients
Hao W,Meng H,Li H,Zheng Y,Song C,Jiang Z,Bai X,Zhang Z,Du L,Liu P,Wu H
Wed Oct 05 00:00:00 UTC 2022
KDM2B overexpression prevents myocardial ischemia-reperfusion injury in rats through regulating inflammatory response via the TLR4/NF-κB p65 axis.
Experimental and therapeutic medicine
Wei Z,Luo L,Hu S,Tian R,Liu Z
Tue Feb 01 00:00:00 UTC 2022
Qing-Re-Xiao-Zheng-Yi-Qi formula relieves kidney damage and activates mitophagy in diabetic kidney disease.
Frontiers in pharmacology
Wu Q,Yan R,Yang H,Wang Y,Zhang C,Zhang J,Cui Z,Wang Y,Sun W
Wed Jan 11 00:00:00 UTC 2023
Homozygous missense mutation in CCDC155 disrupts the transmembrane distribution of CCDC155 and SUN1, resulting in non-obstructive azoospermia and premature ovarian insufficiency in humans.
Human genetics
Wu H,Zhang X,Hua R,Li Y,Cheng L,Li K,Liu Y,Gao Y,Shen Q,Wang G,Lv M,Xu Y,He X,Cao Y,Liu M
Tue Nov 01 00:00:00 UTC 2022
Hepatic inactivation of murine Surf4 results in marked reduction in plasma cholesterol.
eLife
Tang VT,McCormick J,Xu B,Wang Y,Fang H,Wang X,Siemieniak D,Khoriaty R,Emmer BT,Chen XW,Ginsburg D
Tue Oct 04 00:00:00 UTC 2022
Gut microbiota-derived metabolites contribute negatively to hindgut barrier function development at the early weaning goat model.
Animal nutrition (Zhongguo xu mu shou yi xue hui)
Zhang K,Xu Y,Yang Y,Guo M,Zhang T,Zong B,Huang S,Suo L,Ma B,Wang X,Wu Y,Brugger D,Chen Y
Thu Sep 01 00:00:00 UTC 2022
The Tobacco β-Cembrenediol: A Prostate Cancer Recurrence Suppressor Lead and Prospective Scaffold via Modulation of Indoleamine 2,3-Dioxygenase and Tryptophan Dioxygenase.
Nutrients
Mudhish EA,Siddique AB,Ebrahim HY,Abdelwahed KS,King JA,El Sayed KA
Mon Apr 04 00:00:00 UTC 2022
NPM1 promotes cell proliferation by targeting PRDX6 in colorectal cancer.
The international journal of biochemistry & cell biology
Wang D,Li Y,Liu Y,Cheng S,Liu F,Zuo R,Ding C,Shi S,Liu G
Wed Jun 01 00:00:00 UTC 2022
Increased expression of osteopontin in subchondral bone promotes bone turnover and remodeling, and accelerates the progression of OA in a mouse model.
Aging
Lin C,Chen Z,Guo D,Zhou L,Lin S,Li C,Li S,Wang X,Lin B,Ding Y
Tue Jan 04 00:00:00 UTC 2022
PVT1/miR-145-5p/HK2 modulates vascular smooth muscle cells phenotype switch via glycolysis: The new perspective on the spiral artery remodeling.
Placenta
Wu M,Liu W,Huang H,Chen Z,Chen Y,Zhong Y,Jin Z,Liu X,Zou L
Thu Dec 01 00:00:00 UTC 2022
Knockdown of TTC9 inhibits the proliferation, migration and invasion, but induces the apoptosis of lung adenocarcinoma cells.
Heliyon
Huang X,Jiang L,Wen Z,Yuan M,Zhong Y
Tue Nov 01 00:00:00 UTC 2022
The phytochemical constituents and protective effect of Fritillaria hupehensis on acute lung injury.
Fitoterapia
Xiang ML,Zhao YL,Liu YY,Yan XJ,Chen S,Luo XD
Sat Oct 01 00:00:00 UTC 2022
Discovery of a novel Pleuromutilin derivative as anti-IPF lead compound via high-throughput assay.
European journal of medicinal chemistry
Zhang K,Liang J,Wang N,Li N,Jiang Y,Li X,Yang C,Zhou H,Yang G
Sat Nov 05 00:00:00 UTC 2022
Cognitive enhancement and neuroprotective effects of OABL, a sesquiterpene lactone in 5xFAD Alzheimer's disease mice model.
Redox biology
Tang JJ,Huang LF,Deng JL,Wang YM,Guo C,Peng XN,Liu Z,Gao JM
Fri Apr 01 00:00:00 UTC 2022
The Synergistic Cytotoxic Effects of GW5074 and Sorafenib by Impacting Mitochondrial Functions in Human Colorectal Cancer Cell Lines.
Frontiers in oncology
Hu JM,Chang YL,Hsieh CC,Huang SM
Sat Jul 16 00:00:00 UTC 2022
Repeated Yueju, But Not Fluoxetine, Induced Sustained Antidepressant Activity in a Mouse Model of Chronic Learned Helplessness: Involvement of CaMKII Signaling in the Hippocampus.
Evidence-based complementary and alternative medicine : eCAM
Zou Z,Huang J,Yang Q,Zhang Y,Xu B,Wang P,Chen G
Mon May 02 00:00:00 UTC 2022
Hypoxia-Regulated lncRNA USP2-AS1 Drives Head and Neck Squamous Cell Carcinoma Progression.
Cells
Tang J,Wu Z,Wang X,Hou Y,Bai Y,Tian Y
Fri Oct 28 00:00:00 UTC 2022
Sperm Autoantigenic Protein 17 Predicts the Prognosis and the Immunotherapy Response of Cancers: A Pan-Cancer Analysis.
Frontiers in immunology
Tu Z,Peng J,Long X,Li J,Wu L,Huang K,Zhu X
Mon May 23 00:00:00 UTC 2022
Age-related visual impairments and retinal ganglion cells axonal degeneration in a mouse model harboring OPTN (E50K) mutation.
Cell death & disease
Hou M,Shao Z,Zhang S,Liu X,Fan P,Jiang M,Zhao Y,Xiao R,Yuan H
Mon Apr 18 00:00:00 UTC 2022
Identification of endocrine-disrupting chemicals targeting the genes and pathways of genital anomalies in males.
Ecotoxicology and environmental safety
Zhou X,Zhang X,Zhou X,Abulimiti G,Wang Y,Zhang Q,Cong R,Ji C,Luan J,Yao L,Yang J,Song N
Thu Dec 01 00:00:00 UTC 2022
Analgesic effect of nobiletin against neuropathic pain induced by the chronic constriction injury of the sciatic nerve in mice.
Phytotherapy research : PTR
Zhu C,Tian M,Liu N,Ma L,Lan X,Yang J,Du J,Ma H,Li Y,Zheng P,Yu J,Peng X
Thu Sep 01 00:00:00 UTC 2022
Inhibition of base editors with anti-deaminases derived from viruses.
Nature communications
Liu Z,Chen S,Lai L,Li Z
Tue Feb 01 00:00:00 UTC 2022
BRD7 Stabilizes P53 via Dephosphorylation of MDM2 to Inhibit Tumor Growth in Breast Cancer Harboring Wild-type P53.
Journal of Cancer
Luo Y,Wang X,Niu W,Zhou Y,Li M,Ma J,Yang J,Fan S,Zeng Z,Xiong W,Li X,Li G,Xiao J,Zhou M
Tue Apr 05 00:00:00 UTC 2022
Prrx1 promotes resistance to temozolomide by upregulating ABCC1 and inducing vasculogenic mimicry in glioma.
American journal of cancer research
Chen Z,Zhang Y,Chen Y,Lin W,Zhang Y,Cai G,Sun X,Zheng K,He J,Ai T,Wang J,Zhao L,Ke Y
Tue Sep 20 00:00:00 UTC 2022
Protective Effects of Scutellarin on Acute Alcohol Intestinal Injury.
Chemistry & biodiversity
Liu F,Gao X,Li Z,Zhang X,Fan H,Yu G,Bello BK,Feng X,Li D,Teng D,Chen Y,Zhao P,Fu M,Dong J
Fri Apr 01 00:00:00 UTC 2022
Small RNA perspective of physical exercise-related improvement of male reproductive dysfunction due to obesity.
Frontiers in endocrinology
Lin T,Zhang S,Zhou Y,Wu L,Liu X,Huang H
Tue Dec 20 00:00:00 UTC 2022
Diprotin A TFA Exerts Neurovascular Protection in Ischemic Cerebral Stroke.
Frontiers in neuroscience
Zhou MY,Zhang YJ,Ding HM,Wu WF,Cai WW,Wang YQ,Geng DQ
Sat Jul 16 00:00:00 UTC 2022
Z. morio Hemolymph Relieves E. coli-Induced Mastitis by Inhibiting Inflammatory Response and Repairing the Blood-Milk Barrier.
International journal of molecular sciences
Zou Y,Wang X,Xu J,Wang S,Li S,Zhu Y,Wang J
Mon Oct 31 00:00:00 UTC 2022
Supervillin Contributes to LPS-induced Inflammatory Response in THP-1 Cell-derived Macrophages.
Inflammation
Zhou J,Que Y,Pan L,Li X,Zhu C,Jin L,Li S
Tue Feb 01 00:00:00 UTC 2022
UFL1 alleviates ER stress and apoptosis stimulated by LPS via blocking the ferroptosis pathway in human granulosa-like cells.
Cell stress & chaperones
Li J,Tang X,Tu X,Jin Z,Dong H,Yang Q,Yao T,Pan Z
Thu Sep 01 00:00:00 UTC 2022
Proteome-wide analysis of the hippocampus in adolescent male mice with learning and memory impairment caused by chronic ethanol exposure.
Neurobiology of learning and memory
Liu D,Liu S,Li J,Liu X,Wu X,Peng Y,Shen Q
Sat Oct 01 00:00:00 UTC 2022
Effect of Silkworm Pupa Protein Hydrolysates on Proliferation of Gastric Cancer Cells In Vitro.
Foods (Basel, Switzerland)
Li W,Mu L,Zou Y,Wang W,Zhao H,Wu X,Liao S
Sun Aug 07 00:00:00 UTC 2022
Dexmedetomidine Can Enhance PINK1/Parkin-Mediated Mitophagy in MPTP-Induced PD Mice Model by Activating AMPK.
Oxidative medicine and cellular longevity
Chen C,Chen Y,Liu T,Song D,Ma D,Cheng O
Tue May 10 00:00:00 UTC 2022
MiR-3571 modulates the proliferation and migration of vascular smooth muscle cells by targeting claudin 1.
International journal of medical sciences
Xie Y,Tan J,Qin Y,Cao Y,Wang Y,Li A,Wang Z,Qiao Z,Yan Z
Tue Apr 05 00:00:00 UTC 2022
Nrf2 loss of function exacerbates endoplasmic reticulum stress-induced apoptosis in TBI mice.
Neuroscience letters
Sun G,Zhao Z,Lang J,Sun B,Zhao Q
Sun Jan 23 00:00:00 UTC 2022
Extracellular vesicles derived from human umbilical cord mesenchymal stem cells alleviate osteoarthritis of the knee in mice model by interacting with METTL3 to reduce m6A of NLRP3 in macrophage.
Stem cell research & therapy
Zhou H,Shen X,Yan C,Xiong W,Ma Z,Tan Z,Wang J,Li Y,Liu J,Duan A,Liu F
Sat Jul 16 00:00:00 UTC 2022
The Exocyst Is Required for CD36 Fatty Acid Translocase Trafficking and Free Fatty Acid Uptake in Skeletal Muscle Cells.
Cells
Nakamura NK,Tokunaga DS,Ha HY,Polgar N
Sat Aug 06 00:00:00 UTC 2022
AQP5 Is a Novel Prognostic Biomarker in Pancreatic Adenocarcinoma.
Frontiers in oncology
Chen G,Song H,Yang Z,Du T,Zheng Y,Lu Z,Zhang K,Wei D
Sun Nov 05 00:00:00 UTC 2023
Avasimibe Alleviates Disruption of the Airway Epithelial Barrier by Suppressing the Wnt/β-Catenin Signaling Pathway.
Frontiers in pharmacology
Zhou Z,Liang S,Zhou Z,Liu J,Meng X,Zou F,Yu C,Cai S
Tue Mar 01 00:00:00 UTC 2022
Aperture Modulation of Isoreticular Metal Organic Frameworks for Targeted Antitumor Drug Delivery.
ACS applied materials & interfaces
Cai M,Liang W,Wang K,Yin D,Fu T,Zhu R,Qu C,Dong X,Ni J,Yin X
Wed Aug 17 00:00:00 UTC 2022
Functional Attenuation of UCP1 as the Potential Mechanism for a Thickened Blubber Layer in Cetaceans.
Molecular biology and evolution
Zhou M,Wu T,Chen Y,Xu S,Yang G
Thu Nov 03 00:00:00 UTC 2022
The roles of grouper TAK1 in regulating the infection of Singapore grouper iridovirus.
Fish & shellfish immunology
Zhang L,Kang S,Chen H,Liao J,Sun M,Wu S,Xu Z,Xu L,Zhang X,Qin Q,Wei J
Sun May 01 00:00:00 UTC 2022
Downregulation of TUSC3 promotes EMT and hepatocellular carcinoma progression through LIPC/AKT axis.
Journal of translational medicine
Deng R,Lu X,Hong C,Cai R,Wang P,Xiong L,Wang X,Chen Q,Lin J
Sun Oct 23 00:00:00 UTC 2022
Transient Receptor Potential Cation Channel Subfamily V Member 1 Expression Promotes Chemoresistance in Non-Small-Cell Lung Cancer.
Frontiers in oncology
Li L,Chen C,Xiang Q,Fan S,Xiao T,Chen Y,Zheng D
Wed Apr 13 00:00:00 UTC 2022
A synthetic pregnenolone analog promotes microtubule dynamics and neural development.
Cell & bioscience
Kolas V,Bandonil JSA,Wali N,Hsia KC,Shie JJ,Chung BC
Thu Dec 01 00:00:00 UTC 2022
Targeting PERK mediated endoplasmic reticulum stress attenuates neuroinflammation and alleviates lipopolysaccharide-induced depressive-like behavior in male mice.
International immunopharmacology
Xu XF,Shi MM,Luo MY,Liu DD,Guo DM,Ling C,Zhong XL,Xu Y,Cao WY
Sat Oct 01 00:00:00 UTC 2022
Oar-miR-432 Regulates Fat Differentiation and Promotes the Expression of BMP2 in Ovine Preadipocytes.
Frontiers in genetics
Jin M,Fei X,Li T,Lu Z,Chu M,Di R,He X,Wang X,Wang Y,Yuan Z,Quan K,Wang H,Wei C
Sat Jul 16 00:00:00 UTC 2022
Hydrogen Sulfide Regulates SERCA2a Ubiquitylation via Muscle RING Finger-1 S-Sulfhydration to Affect Cardiac Contractility in db/db Mice.
Cells
Peng S,Zhao D,Li Q,Wang M,Zhang S,Pang K,Huang J,Lu F,Chen H,Zhang W
Wed Nov 02 00:00:00 UTC 2022
Reticulocalbin3: A Ca2+ homeostasis regulator that promotes esophageal squamous cell carcinoma progression and cisplatin resistance.
Cancer science
Cai R,Wang P,Zhao X,Lu X,Deng R,Wang X,Hong C,Lin J
Sat Oct 01 00:00:00 UTC 2022
Amelioration of scopolamine-induced learning and memory impairment by the TRPV4 inhibitor HC067047 in ICR mice.
Neuroscience letters
Deng Y,Li W,Niu L,Luo X,Li J,Zhang Y,Liu H,He J,Wan W
Mon Jan 10 00:00:00 UTC 2022
Time series RNA-seq analysis identifies MAPK10 as a critical gene in diabetes mellitus-induced atrial fibrillation in mice.
Journal of molecular and cellular cardiology
Liu F,Deng Y,Zhao Y,Li Z,Gao J,Zhang Y,Yang X,Liu Y,Xia Y
Fri Jul 01 00:00:00 UTC 2022
ETV5 Regulates Hepatic Fatty Acid Metabolism Through PPAR Signaling Pathway.
Diabetes
Mao Z,Feng M,Li Z,Zhou M,Xu L,Pan K,Wang S,Su W,Zhang W
Fri Jan 01 00:00:00 UTC 2021
MICAL2PV suppresses the formation of tunneling nanotubes and modulates mitochondrial trafficking.
EMBO reports
Wang F,Chen X,Cheng H,Song L,Liu J,Caplan S,Zhu L,Wu JY
Mon Jul 05 00:00:00 UTC 2021
MICAL2PV suppresses the formation of tunneling nanotubes and modulates mitochondrial trafficking.
EMBO reports
Wang F,Chen X,Cheng H,Song L,Liu J,Caplan S,Zhu L,Wu JY
Mon Jul 05 00:00:00 UTC 2021
lncRNA Ttc3-209 Promotes the Apoptosis of Retinal Ganglion Cells in Retinal Ischemia Reperfusion Injury by Targeting the miR-484/Wnt8a Axis.
Investigative ophthalmology & visual science
Zhang R,Feng Y,Lu J,Ge Y,Li H
Mon Mar 01 00:00:00 UTC 2021
Inhibition of ERAD synergizes with FTS to eradicate pancreatic cancer cells.
BMC cancer
Du R,Sullivan DK,Azizian NG,Liu Y,Li Y
Sat Mar 06 00:00:00 UTC 2021
Decreased NHE3 activity in intestinal epithelial cells in TGEV and PEDV-induced piglet diarrhea.
Veterinary microbiology
Niu Z,Zhang Y,Kan Z,Ran L,Yan T,Xu S,Zhang S,Zhang J,Zou H,Song Z
Wed Dec 01 00:00:00 UTC 2021
MnSOD Lysine 68 acetylation leads to cisplatin and doxorubicin resistance due to aberrant mitochondrial metabolism.
International journal of biological sciences
Gao Y,Zhu Y,Tran EL,Tokars V,Dean AE,Quan S,Gius D
Thu Mar 24 00:00:00 UTC 2022
HRD1-mediated METTL14 degradation regulates m6A mRNA modification to suppress ER proteotoxic liver disease.
Molecular cell
Wei J,Harada BT,Lu D,Ma R,Gao B,Xu Y,Montauti E,Mani N,Chaudhuri SM,Gregory S,Weinberg SE,Zhang DD,Green R,He C,Fang D
Thu Dec 16 00:00:00 UTC 2021
The Role of KDM2B and EZH2 in Regulating the Stemness in Colorectal Cancer Through the PI3K/AKT Pathway.
Frontiers in oncology
Sanches JGP,Song B,Zhang Q,Cui X,Yabasin IB,Ntim M,Li X,He J,Zhang Y,Mao J,Lu Y,Li L
Fri Apr 02 00:00:00 UTC 2021
Identification and Bioinformatic Assessment of circRNA Expression After RMI1 Knockdown and Ionizing Radiation Exposure.
DNA and cell biology
Sun Y,Fang L,Yang M,He N,Wang J,Zhang M,Ji K,Wang Q,Liu Y,Du L,Wang Y,Xu C,Liu Q
Fri Jan 01 00:00:00 UTC 2021
Arsenic trioxide induces expression of BCL-2 expression via NF-κB and p38 MAPK signaling pathways in BEAS-2B cells during apoptosis.
Ecotoxicology and environmental safety
Tang J,Yao C,Liu Y,Yuan J,Wu L,Hosoi K,Yu S,Huang C,Wei H,Chen G
Fri Oct 01 00:00:00 UTC 2021
KPNB1-mediated nuclear translocation of PD-L1 promotes non-small cell lung cancer cell proliferation via the Gas6/MerTK signaling pathway.
Cell death and differentiation
Du W,Zhu J,Zeng Y,Liu T,Zhang Y,Cai T,Fu Y,Zhang W,Zhang R,Liu Z,Huang JA
Thu Apr 01 00:00:00 UTC 2021
Protective Effect of Remdesivir Against Pulmonary Fibrosis in Mice.
Frontiers in pharmacology
Li X,Liu R,Cui Y,Liang J,Bi Z,Li S,Miao Y,Zhang L,Li X,Zhou H,Yang C
Tue Sep 14 00:00:00 UTC 2021
Isoeucommin A attenuates kidney injury in diabetic nephropathy through the Nrf2/HO-1 pathway.
FEBS open bio
Huang Q,Ouyang DS,Liu Q
Sun Aug 01 00:00:00 UTC 2021
Bioinformatic Approach to Unveil Key Differentially Expressed Proteins in Human Sperm After Slow and Rapid Cryopreservation.
Frontiers in cell and developmental biology
Corda PO,Silva JV,Pereira SC,Barros A,Alves MG,Fardilha M
Wed Feb 16 00:00:00 UTC 2022
Salidroside Attenuates Doxorubicin-Induced Cardiac Dysfunction Partially Through Activation of QKI/FoxO1 Pathway.
Journal of cardiovascular translational research
Yan F,Liu R,Zhuang X,Li R,Shi H,Gao X
Thu Apr 01 00:00:00 UTC 2021
The E2F1/USP11 positive feedback loop promotes hepatocellular carcinoma metastasis and inhibits autophagy by activating ERK/mTOR pathway.
Cancer letters
Qiao L,Zhang Q,Sun Z,Liu Q,Wu Z,Hu W,Bao S,Yang Q,Liu L
Sat Aug 28 00:00:00 UTC 2021
PADI6 Regulates Trophoblast Cell Migration-Invasion Through the Hippo/YAP1 Pathway in Hydatidiform Moles.
Journal of inflammation research
Huang B,Zhao Y,Zhou L,Gong T,Feng J,Han P,Qian J
Mon Apr 25 00:00:00 UTC 2022
Gossypol, a novel modulator of VCP, induces autophagic degradation of mutant huntingtin by promoting the formation of VCP/p97-LC3-mHTT complex.
Acta pharmacologica Sinica
Li XJ,Zhang YY,Fu YH,Zhang H,Li HX,Li QF,Li HL,Tan RK,Jiang CX,Jiang W,Li ZX,Luo C,Lu BX,Dang YJ
Fri Oct 01 00:00:00 UTC 2021
Extracellular vesicles derived from LPS-preconditioned human synovial mesenchymal stem cells inhibit extracellular matrix degradation and prevent osteoarthritis of the knee in a mouse model.
Stem cell research & therapy
Duan A,Shen K,Li B,Li C,Zhou H,Kong R,Shao Y,Qin J,Yuan T,Ji J,Guo W,Wang X,Xue T,Li L,Huang X,Sun Y,Cai Z,Liu W,Liu F
Wed Jul 28 00:00:00 UTC 2021
NADPH levels affect cellular epigenetic state by inhibiting HDAC3-Ncor complex.
Nature metabolism
Li W,Kou J,Qin J,Li L,Zhang Z,Pan Y,Xue Y,Du W
Fri Jan 01 00:00:00 UTC 2021
Guaiazulene derivative 1,2,3,4-tetrahydroazuleno[1,2-b] tropone reduces the production of ATP by inhibiting electron transfer complex II.
FEBS open bio
Kasami C,Yamaguchi JI,Inoue H
Mon Nov 01 00:00:00 UTC 2021
Integrated Metabolomics and Transcriptomics Analyses Reveal Metabolic Landscape in Neuronal Cells during JEV Infection.
Virologica Sinica
Li M,Yang J,Ye C,Bian P,Yang X,Zhang H,Luo C,Xue Z,Lei Y,Lian J
Wed Dec 01 00:00:00 UTC 2021
Cypermethrin triggers YY1-mediated testosterone biosynthesis suppression.
Ecotoxicology and environmental safety
Liu C,Qu J,Wu M,Huang X,Li L
Wed Dec 01 00:00:00 UTC 2021
Triptolide impairs glycolysis by suppressing GATA4/Sp1/PFKP signaling axis in mouse Sertoli cells.
Toxicology and applied pharmacology
Zhang Y,Tang Y,Luo Y,Luo L,Shen F,Huang Z
Sun Aug 15 00:00:00 UTC 2021
Hyper-SUMOylation of SMN induced by SENP2 deficiency decreases its stability and leads to spinal muscular atrophy-like pathology.
Journal of molecular medicine (Berlin, Germany)
Zhang Y,Chen X,Wang Q,Du C,Lu W,Yuan H,Zhang Z,Li D,Ling X,Ren X,Zhao Y,Su Q,Xing Z,Qin Y,Yang X,Shen Y,Wu H,Qi Y
Wed Dec 01 00:00:00 UTC 2021
Attractin Gene Deficiency in Rats Leads to Impairments in Both Activity and Spatial Learning and Memory.
Neuroscience
Li XH,Xue C,Liu MQ,Zhang MY,Zhou Y,Xiao X,Wang J,Xu XJ,Shi Y,Zhang WN
Thu Jul 01 00:00:00 UTC 2021
TRIM21-regulated Annexin A2 plasma membrane trafficking facilitates osteosarcoma cell differentiation through the TFEB-mediated autophagy.
Cell death & disease
Zhang HT,Zeng Q,Wu B,Lu J,Tong KL,Lin J,Liu QY,Xu L,Yang J,Liu X,Liu W,Zhang YF,Lian Q,Liu L,Gao X
Wed Jan 06 00:00:00 UTC 2021
Umbilical Cord Mesenchymal Stem Cell-Derived Exosomes Ameliorate HaCaT Cell Photo-Aging.
Rejuvenation research
Liu SJ,Meng MY,Han S,Gao H,Zhao YY,Yang Y,Lin ZY,Yang LR,Zhu K,Han R,Huang WW,Wang RQ,Yang LL,Wang WJ,Li L,Wang XD,Hou ZL,Liao LW,Yang L
Sun Aug 01 00:00:00 UTC 2021
The role of SAMM50 in non-alcoholic fatty liver disease: from genetics to mechanisms.
FEBS open bio
Li Z,Shen W,Wu G,Qin C,Zhang Y,Wang Y,Song G,Xiao C,Zhang X,Deng G,Wang R,Wang X
Thu Jul 01 00:00:00 UTC 2021
Neuronal extracellular vesicle derived miR-98 prevents salvageable neurons from microglial phagocytosis in acute ischemic stroke.
Cell death & disease
Yang J,Cao LL,Wang XP,Guo W,Guo RB,Sun YQ,Xue TF,Cai ZY,Ji J,Cheng H,Sun XL
Wed Jan 06 00:00:00 UTC 2021
Expression and prognostic potential of PLEK2 in head and neck squamous cell carcinoma based on bioinformatics analysis.
Cancer medicine
Wang J,Sun Z,Wang J,Tian Q,Huang R,Wang H,Wang X,Han F
Wed Sep 01 00:00:00 UTC 2021
MicroRNA‑8063 targets heterogeneous nuclear ribonucleoprotein AB to inhibit the self‑renewal of colorectal cancer stem cells via the Wnt/β‑catenin pathway.
Oncology reports
Chen ZQ,Yuan T,Jiang H,Yang YY,Wang L,Fu RM,Luo SQ,Zhang T,Wu ZY,Wen KM
Fri Oct 01 00:00:00 UTC 2021
The Regulatory Role of GBF1 on Osteoclast Activation Through EIF2a Mediated ER Stress and Novel Marker FAM129A Induction.
Frontiers in cell and developmental biology
Wen C,Zhou Y,Xu Y,Tan H,Pang C,Liu H,Liu K,Wei L,Luo H,Qin T,He C,Liu C,Zhou C
Tue Sep 14 00:00:00 UTC 2021
Mechanically Activated Calcium Channel PIEZO1 Modulates Radiation-Induced Epithelial-Mesenchymal Transition by Forming a Positive Feedback With TGF-β1.
Frontiers in molecular biosciences
Huang JQ,Zhang H,Guo XW,Lu Y,Wang SN,Cheng B,Dong SH,Lyu XL,Li FS,Li YW
Tue Nov 02 00:00:00 UTC 2021
UM-6 induces autophagy and apoptosis via the Hippo-YAP signaling pathway in cervical cancer.
Cancer letters
Wang D,He J,Dong J,Wu S,Liu S,Zhu H,Xu T
Thu Oct 28 00:00:00 UTC 2021
Tributyltin Oxide Exposure During in vitro Maturation Disrupts Oocyte Maturation and Subsequent Embryonic Developmental Competence in Pigs.
Frontiers in cell and developmental biology
Xiao Y,Yuan B,Hu W,Qi J,Jiang H,Sun B,Zhang J,Liang S
Fri Jul 16 00:00:00 UTC 2021
Alternative splicing transitions associate with emerging atrophy phenotype during denervation-induced skeletal muscle atrophy.
Journal of cellular physiology
Qiu J,Wu L,Chang Y,Sun H,Sun J
Tue Jun 01 00:00:00 UTC 2021
The STING antagonist H-151 ameliorates psoriasis via suppression of STING/NF-κB-mediated inflammation.
British journal of pharmacology
Pan Y,You Y,Sun L,Sui Q,Liu L,Yuan H,Chen C,Liu J,Wen X,Dai L,Sun H
Wed Dec 01 00:00:00 UTC 2021
A Nuclear Export Signal Is Required for cGAS to Sense Cytosolic DNA.
Cell reports
Sun H,Huang Y,Mei S,Xu F,Liu X,Zhao F,Yin L,Zhang D,Wei L,Wu C,Ma S,Wang J,Cen S,Liang C,Hu S,Guo F
Tue Jan 05 00:00:00 UTC 2021
Cucurbit[7]uril as a Broad-Spectrum Antiviral Agent against Diverse RNA Viruses.
Virologica Sinica
Quan J,Zhang X,Ding Y,Li S,Qiu Y,Wang R,Zhou X
Fri Oct 01 00:00:00 UTC 2021
DCI after Aneurysmal Subarachnoid Hemorrhage Is Related to the Expression of MFG-E8.
BioMed research international
Chen X,Jiang Y,Liu J,Wang C,Wan D,Liang A,Leng J,Yang Y,Xiang H,Liu R
Fri Jan 14 00:00:00 UTC 2022
Ameliorative effects of eosinophil deficiency on immune response, endoplasmic reticulum stress, apoptosis, and autophagy in fungus-induced allergic lung inflammation.
Respiratory research
Wang S,Jiang Z,Li L,Zhang J,Zhang C,Shao C
Mon Jun 07 00:00:00 UTC 2021
Ilexsaponin A1 Ameliorates Diet-Induced Nonalcoholic Fatty Liver Disease by Regulating Bile Acid Metabolism in Mice.
Frontiers in pharmacology
Zhao WW,Xiao M,Wu X,Li XW,Li XX,Zhao T,Yu L,Chen XQ
Sat Jan 01 00:00:00 UTC 2022
PI3K/AKT pathway mediates the antidepressant- and anxiolytic-like roles of hydrogen sulfide in streptozotocin-induced diabetic rats via promoting hippocampal neurogenesis.
Neurotoxicology
Jiang W,Tang YY,Zhu WW,Li C,Zhang P,Li RQ,Chen YJ,Zou W,Tang XQ
Thu Jul 01 00:00:00 UTC 2021
The combination of G-CSF and AMD3100 mobilizes bone marrow-derived stem cells to protect against cisplatin-induced acute kidney injury in mice.
Stem cell research & therapy
Chen Z,Ren X,Ren R,Wang Y,Shang J
Wed Mar 24 00:00:00 UTC 2021
Reduced activity of intestinal surface Na+/H+ exchanger NHE3 is a key factor for induction of diarrhea after PEDV infection in neonatal piglets.
Virology
Song Z,Yan T,Ran L,Niu Z,Zhang Y,Kan Z,Xu S,Zhang S,Zhang J,Zou H,Lei C
Mon Nov 01 00:00:00 UTC 2021
RNF128 suppresses the malignancy of colorectal cancer cells via inhibition of Wnt/β-catenin signaling.
American journal of translational research
Zhu Y,Gan Y,Zou R,Sha H,Lu Y,Zhang Y,Feng J
Tue Jan 18 00:00:00 UTC 2022
Characterization of ALTO-encoding circular RNAs expressed by Merkel cell polyomavirus and trichodysplasia spinulosa polyomavirus.
PLoS pathogens
Yang R,Lee EE,Kim J,Choi JH,Kolitz E,Chen Y,Crewe C,Salisbury NJH,Scherer PE,Cockerell C,Smith TR,Rosen L,Verlinden L,Galloway DA,Buck CB,Feltkamp MC,Sullivan CS,Wang RC
Sat May 01 00:00:00 UTC 2021
Prrx1 promotes stemness and angiogenesis via activating TGF-β/smad pathway and upregulating proangiogenic factors in glioma.
Cell death & disease
Chen Z,Chen Y,Li Y,Lian W,Zheng K,Zhang Y,Zhang Y,Lin C,Liu C,Sun F,Sun X,Wang J,Zhao L,Ke Y
Tue Jun 15 00:00:00 UTC 2021
Loss of Diacylglycerol Kinase α Enhances Macrophage Responsiveness.
Frontiers in immunology
Manigat LC,Granade ME,Taori S,Miller CA,Vass LR,Zhong XP,Harris TE,Purow BW
Fri Jan 07 00:00:00 UTC 2022
GluN2A/ERK/CREB Signaling Pathway Involved in Electroacupuncture Regulating Hypothalamic-Pituitary-Adrenal Axis Hyperactivity.
Frontiers in neuroscience
Wang Y,Han J,Zhu J,Zhang M,Ju M,Du Y,Tian Z
Fri Oct 22 00:00:00 UTC 2021
Betulinic acid attenuated bleomycin-induced pulmonary fibrosis by effectively intervening Wnt/β-catenin signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li X,Liu X,Deng R,Gao S,Jiang Q,Liu R,Li H,Miao Y,Zhai Y,Zhang S,Wang Z,Ren Y,Ning W,Zhou H,Yang C
Fri Jan 01 00:00:00 UTC 2021
PDGF/MEK/ERK axis represses Ca2+ clearance via decreasing the abundance of plasma membrane Ca2+ pump PMCA4 in pulmonary arterial smooth muscle cells.
American journal of physiology. Cell physiology
Deng L,Chen J,Wang T,Chen B,Yang L,Liao J,Chen Y,Wang J,Tang H,Yi J,Kang K,Li L,Gou D
Fri Jan 01 00:00:00 UTC 2021
Latent Sex Differences in CaMKII-nNOS Signaling That Underlie Antidepressant-Like Effects of Yueju-Ganmaidazao Decoction in the Hippocampus.
Frontiers in behavioral neuroscience
Yin Y,Qian S,Chen Y,Sun Y,Li Y,Yu Y,Li J,Wu Z,Yu X,Ge R,Han J,Sun D,Wu H,Liu L,Xue W,Wang W
Sat Jul 24 00:00:00 UTC 2021
Melatonin ameliorates cypermethrin-induced impairments by regulating oxidative stress, DNA damage and apoptosis in porcine Sertoli cells.
Theriogenology
Li J,Sun BX,Wang DL,Liu Y,Qi JJ,Nie XW,Bai CY,Zhang JB,Liang S
Tue Jun 01 00:00:00 UTC 2021
Human umbilical cord-derived mesenchymal stem cell therapy ameliorates lupus through increasing CD4+ T cell senescence via MiR-199a-5p/Sirt1/p53 axis.
Theranostics
Cheng T,Ding S,Liu S,Li Y,Sun L
Wed Aug 04 00:00:00 UTC 2021
Tripartite motif-containing protein 16 protects against myocardial ischemia/reperfusion injury by affecting the Keap1/Nrf2 axis.
Cell and tissue research
Cui Q,Yan L
Mon Nov 01 00:00:00 UTC 2021
Iron Overload-Induced Ferroptosis Impairs Porcine Oocyte Maturation and Subsequent Embryonic Developmental Competence in vitro.
Frontiers in cell and developmental biology
Hu W,Zhang Y,Wang D,Yang T,Qi J,Zhang Y,Jiang H,Zhang J,Sun B,Liang S
Tue Jun 15 00:00:00 UTC 2021
Nuclear iASPP determines cell fate by selectively inhibiting either p53 or NF-κB.
Cell death discovery
Ge W,Wang Y,Zheng S,Zhao D,Wang X,Zhang X,Hu Y
Mon Jul 26 00:00:00 UTC 2021
Formaldehyde induces ferroptosis in hippocampal neuronal cells by upregulation of the Warburg effect.
Toxicology
Li XN,Yang SQ,Li M,Li XS,Tian Q,Xiao F,Tang YY,Kang X,Wang CY,Zou W,Zhang P,Tang XQ
Sat Jan 30 00:00:00 UTC 2021
The m6A Reader YTHDF1 Facilitates the Tumorigenesis and Metastasis of Gastric Cancer via USP14 Translation in an m6A-Dependent Manner.
Frontiers in cell and developmental biology
Chen XY,Liang R,Yi YC,Fan HN,Chen M,Zhang J,Zhu JS
Thu Apr 21 00:00:00 UTC 2022
Expression and Functional Analysis of lncRNAs Involved in Platelet-Derived Growth Factor-BB-Induced Proliferation of Human Aortic Smooth Muscle Cells.
Frontiers in cardiovascular medicine
Lin JJ,Chen W,Gong M,Xu X,Du MY,Wang SF,Yang LY,Wang Y,Liu KX,Kong P,Li B,Liu K,Li YM,Dong LH,Sun SG
Sat Sep 25 00:00:00 UTC 2021
Libertellenone H, a Natural Pimarane Diterpenoid, Inhibits Thioredoxin System and Induces ROS-Mediated Apoptosis in Human Pancreatic Cancer Cells.
Molecules (Basel, Switzerland)
Zhang W,Zhu Y,Yu H,Liu X,Jiao B,Lu X
Sat Jan 09 00:00:00 UTC 2021
CHIP protects against MPP+/MPTP-induced damage by regulating Drp1 in two models of Parkinson's disease.
Aging
Hu Z,Mao C,Wang H,Zhang Z,Zhang S,Luo H,Tang M,Yang J,Yuan Y,Wang Y,Liu Y,Fan L,Zhang Q,Yao D,Liu F,Schisler JC,Shi C,Xu Y
Sat Jan 02 00:00:00 UTC 2021
HSP/HSC70 activity is required for Bombyx mori nucleopolyhedrovirus replication at the early infectious phase.
Microbial pathogenesis
Mao F,Zhu Y,Gao X,Chen X,Ngowo J,Miao M,Quan Y,Yu W
Thu Apr 01 00:00:00 UTC 2021
CXCL5 Downregulation in Villous Tissue Is Correlated With Recurrent Spontaneous Abortion.
Frontiers in immunology
Zhang S,Ding J,Wang J,Yin T,Zhang Y,Yang J
Mon Dec 20 00:00:00 UTC 2021
Glycine alleviates fluoride-induced oxidative stress, apoptosis and senescence in a porcine testicular Sertoli cell line.
Reproduction in domestic animals = Zuchthygiene
Liu Y,Sun B,Zhang S,Li J,Qi J,Bai C,Zhang J,Liang S
Tue Jun 01 00:00:00 UTC 2021
Iptakalim alleviates synaptic damages via targeting mitochondrial ATP-sensitive potassium channel in depression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Guo W,Tang ZY,Cai ZY,Zhao WE,Yang J,Wang XP,Ji J,Huang XX,Sun XL
Sat May 01 00:00:00 UTC 2021
Frugoside delays osteoarthritis progression via inhibiting miR-155-modulated synovial macrophage M1 polarization.
Rheumatology (Oxford, England)
Wang H,Zhang H,Fan K,Zhang D,Hu A,Zeng X,Liu YL,Tan G,Wang H
Sat Oct 02 00:00:00 UTC 2021
TRPV4 inhibitor HC067047 produces antidepressant-like effect in LPS-induced depression mouse model.
Neuropharmacology
Li W,Xu Y,Liu Z,Shi M,Zhang Y,Deng Y,Zhong X,Chen L,He J,Zeng J,Luo M,Cao W,Wan W
Wed Dec 15 00:00:00 UTC 2021
TIPE2 is a checkpoint of natural killer cell maturation and antitumor immunity.
Science advances
Bi J,Cheng C,Zheng C,Huang C,Zheng X,Wan X,Chen YH,Tian Z,Sun H
Fri Sep 17 00:00:00 UTC 2021
BCI Suppresses RANKL-Mediated Osteoclastogenesis and Alleviates Ovariectomy-Induced Bone Loss.
Frontiers in pharmacology
Cai C,Hu W,Zhang Y,Hu X,Yang S,Qiu H,Wang R,Ma M,Qiu Y,Chu T
Tue Nov 23 00:00:00 UTC 2021
Theaflavin Promotes Mitochondrial Abundance and Glucose Absorption in Myotubes by Activating the CaMKK2-AMPK Signal Axis via Calcium-Ion Influx.
Journal of agricultural and food chemistry
Qu Z,Liu A,Liu C,Tang Q,Zhan L,Xiao W,Huang J,Liu Z,Zhang S
Wed Jul 28 00:00:00 UTC 2021
NOD2 deficiency confers a pro-tumorigenic macrophage phenotype to promote lung adenocarcinoma progression.
Journal of cellular and molecular medicine
Wang Y,Miao Z,Qin X,Li B,Han Y
Sun Aug 01 00:00:00 UTC 2021
NORHA, a novel follicular atresia-related lncRNA, promotes porcine granulosa cell apoptosis via the miR-183-96-182 cluster and FoxO1 axis.
Journal of animal science and biotechnology
Yao W,Pan Z,Du X,Zhang J,Liu H,Li Q
Thu Oct 07 00:00:00 UTC 2021
Rosuvastatin Nanomicelles Target Neuroinflammation and Improve Neurological Deficit in a Mouse Model of Intracerebral Hemorrhage.
International journal of nanomedicine
Zi L,Zhou W,Xu J,Li J,Li N,Xu J,You C,Wang C,Tian M
Tue Jun 01 00:00:00 UTC 2021
USP37 downregulation elevates the Chemical Sensitivity of Human Breast Cancer Cells to Adriamycin.
International journal of medical sciences
Qin T,Cui XY,Xiu H,Huang C,Sun ZN,Xu XM,Li LH,Yue L
Mon Sep 13 00:00:00 UTC 2021
Citrus Alkaline Extracts Inhibit Senescence of A549 Cells to Alleviate Pulmonary Fibrosis via the β-Catenin/P53 Pathway.
Medical science monitor : international medical journal of experimental and clinical research
Han D,Xu Y,Peng WP,Feng F,Wang Z,Gu C,Zhou X
Fri Mar 12 00:00:00 UTC 2021
Comparative Proteomics of Rat Olfactory Bulb Reveal Insights into Susceptibility and Resiliency to Chronic-stress-induced Depression or Anxiety.
Neuroscience
Liu D,Cai X,Wang L,Yi F,Liao W,Huang R,Fang C,Chen J,Zhou J
Fri Oct 01 00:00:00 UTC 2021
circCDYL2, Overexpressed in Highly Migratory Colorectal Cancer Cells, Promotes Migration by Binding to Ezrin.
Frontiers in oncology
Li X,Wang J,Long H,Lin W,Wang H,Chen Y,Yuan Q,Li X
Tue Apr 26 00:00:00 UTC 2022
Oral Administration of Probiotic Bifidobacterium breve Improves Facilitation of Hippocampal Memory Extinction via Restoration of Aberrant Higher Induction of Neuropsin in an MPTP-Induced Mouse Model of Parkinson's Disease.
Biomedicines
Ishii T,Furuoka H,Kaya M,Kuhara T
Mon Feb 08 00:00:00 UTC 2021
Therapy of spinal cord injury by zinc modified gold nanoclusters via immune-suppressing strategies.
Journal of nanobiotechnology
Lin S,Li D,Zhou Z,Xu C,Mei X,Tian H
Mon Sep 20 00:00:00 UTC 2021
Naringin Promotes Skeletal Muscle Fiber Remodeling by the AdipoR1-APPL1-AMPK Signaling Pathway.
Journal of agricultural and food chemistry
Li P,Zhang S,Song H,Traore SS,Li J,Raubenheimer D,Cui Z,Kou G
Wed Oct 13 00:00:00 UTC 2021
Cypermethrin induces Sertoli cell apoptosis through mitochondrial pathway associated with calcium.
Toxicology research
Wang HX,Zhang R,Li Z,Wang LS,Yu Y,Wang Q,Ding Z,Zhang JP,Zhang MR,Xu LC
Sun Aug 01 00:00:00 UTC 2021
1,10-Seco-Eudesmane sesquiterpenoids as a new type of anti-neuroinflammatory agents by suppressing TLR4/NF-κB/MAPK pathways.
European journal of medicinal chemistry
Tang JJ,Wang MR,Dong S,Huang LF,He QR,Gao JM
Mon Nov 15 00:00:00 UTC 2021
High-Carbohydrate Diet Alleviates the Oxidative Stress, Inflammation and Apoptosis of Megalobrama amblycephala Following Dietary Exposure to Silver Nanoparticles.
Antioxidants (Basel, Switzerland)
Chen F,Zhao CY,Guan JF,Liu XC,Li XF,Xie DZ,Xu C
Wed Aug 25 00:00:00 UTC 2021
Yixin-Fumai granules improve sick sinus syndrome in aging mice through Nrf-2/HO-1 pathway: A new target for sick sinus syndrome.
Journal of ethnopharmacology
Zhang H,Li L,Hao M,Chen K,Lu Y,Qi J,Chen W,Ren L,Cai X,Chen C,Liu Z,Zhao B,Li Z,Hou P
Wed Sep 15 00:00:00 UTC 2021
The Downregulation of eIF3a Contributes to Vemurafenib Resistance in Melanoma by Activating ERK via PPP2R1B.
Frontiers in pharmacology
Jiang SL,Wang ZB,Zhu T,Jiang T,Fei JF,Liu C,Luo C,Cheng Y,Liu ZQ
Tue Sep 14 00:00:00 UTC 2021
Mbd2 deficiency alleviates retinal cell apoptosisvia the miR-345-5p/Atf1 axis in high glucoseinjury and streptozotocin-induced diabetic mice.
Molecular therapy. Nucleic acids
Ge Y,Zhang R,Feng Y,Lu J,Li H
Fri Dec 03 00:00:00 UTC 2021
AEG-1 Knockdown Sensitizes Glioma Cells to Radiation Through Impairing Homologous Recombination Via Targeting RFC5.
DNA and cell biology
Zhao X,Sun Y,Sun X,Li J,Shi X,Liang Z,Ma Y,Zhang X
Thu Jul 01 00:00:00 UTC 2021
Protocol for nuclear export signal characterization of cGAS in mammalian cells.
STAR protocols
Huang Y,McLean M,Liang C,Guo F
Fri Sep 17 00:00:00 UTC 2021
Inhibition of the NLRP3 inflammasome with MCC950 prevents chronic social isolation-induced depression-like behavior in male mice.
Neuroscience letters
Li W,Niu L,Liu Z,Xu X,Shi M,Zhang Y,Deng Y,He J,Xu Y,Wan W,Sun Q,Zhong X,Cao W
Sat Nov 20 00:00:00 UTC 2021
Extracellular vesicles-based pre-targeting strategy enables multi-modal imaging of orthotopic colon cancer and image-guided surgery.
Journal of nanobiotechnology
Jing B,Qian R,Jiang D,Gai Y,Liu Z,Guo F,Ren S,Gao Y,Lan X,An R
Sat May 22 00:00:00 UTC 2021
ZBP1-MLKL necroptotic signaling potentiates radiation-induced antitumor immunity via intratumoral STING pathway activation.
Science advances
Yang Y,Wu M,Cao D,Yang C,Jin J,Wu L,Hong X,Li W,Lu L,Li J,Wang X,Meng X,Zhang Z,Cheng J,Ye Y,Xiao H,Yu J,Deng L
Fri Oct 08 00:00:00 UTC 2021
A novel tonicity-responsive microRNA miR-23a-5p modulates renal cell survival under osmotic stress through targeting heat shock protein 70 HSPA1B.
American journal of physiology. Cell physiology
Kang K,Liao X,Li Q,Chen J,Niu Y,Zeng Y,Xia S,Zeng L,Liu S,Gou D
Mon Feb 01 00:00:00 UTC 2021
Hypoxia-inducible factor-1α promotes proliferation of airway smooth muscle cells through miRNA-103-mediated signaling pathway under hypoxia.
In vitro cellular & developmental biology. Animal
Zhang C,Gao J,Zhu S
Wed Dec 01 00:00:00 UTC 2021
TMEM41B acts as an ER scramblase required for lipoprotein biogenesis and lipid homeostasis.
Cell metabolism
Huang D,Xu B,Liu L,Wu L,Zhu Y,Ghanbarpour A,Wang Y,Chen FJ,Lyu J,Hu Y,Kang Y,Zhou W,Wang X,Ding W,Li X,Jiang Z,Chen J,Zhang X,Zhou H,Li JZ,Guo C,Zheng W,Zhang X,Li P,Melia T,Reinisch K,Chen XW
Tue Aug 03 00:00:00 UTC 2021
Transcriptome Differences in Pig Tracheal Epithelial Cells in Response to Pasteurella Multocida Infection.
Frontiers in veterinary science
Wang X,Wang F,Lin L,Liang W,Liu S,Hua L,Wang X,Chen H,Peng Z,Wu B
Wed Sep 08 00:00:00 UTC 2021
Pharmacological effects of indole alkaloids from Alstonia scholaris (L.) R. Br. on pulmonary fibrosis in vivo.
Journal of ethnopharmacology
Zhao YL,Pu SB,Qi Y,Wu BF,Shang JH,Liu YP,Hu D,Luo XD
Mon Mar 01 00:00:00 UTC 2021
Sesamin induces cell cycle arrest and apoptosis through p38/C-Jun N-terminal kinase mitogen-activated protein kinase pathways in human colorectal cancer cells.
Anti-cancer drugs
Wang X,Qiao J,Zou C,Zhao Y,Huang Y
Mon Mar 01 00:00:00 UTC 2021
Vagus nerve stimulation mediates microglia M1/2 polarization via inhibition of TLR4 pathway after ischemic stroke.
Biochemical and biophysical research communications
Zhang L,Liu Y,Wang S,Long L,Zang Q,Ma J,Yu L,Jia G
Fri Nov 05 00:00:00 UTC 2021
A network pharmacology approach to reveal the pharmacological targets and biological mechanism of compound kushen injection for treating pancreatic cancer based on WGCNA and in vitro experiment validation.
Chinese medicine
Wu C,Huang ZH,Meng ZQ,Fan XT,Lu S,Tan YY,You LM,Huang JQ,Stalin A,Ye PZ,Wu ZS,Zhang JY,Liu XK,Zhou W,Zhang XM,Wu JR
Mon Nov 22 00:00:00 UTC 2021
BPA modulates the WDR5/TET2 complex to regulate ERβ expression in eutopic endometrium and drives the development of endometriosis.
Environmental pollution (Barking, Essex : 1987)
Xue W,Yao X,Ting G,Ling J,Huimin L,Yuan Q,Chun Z,Ming Z,Yuanzhen Z
Fri Jan 01 00:00:00 UTC 2021
Functional domains and amino acid residues of Japanese eel IRF1, AjIRF1, regulate its nuclear import and IFN/Mx promoter activation.
Developmental and comparative immunology
Li W,Ling L,Wang Z,Liang Y,Huang W,Nie P,Huang B
Mon Mar 01 00:00:00 UTC 2021
EGFR and ERK activation resists flavonoid quercetin-induced anticancer activities in human cervical cancer cells in vitro.
Oncology letters
Chen X,Xu P,Zhang H,Su X,Guo L,Zhou X,Wang J,Huang P,Zhang Q,Sun R
Mon Nov 01 00:00:00 UTC 2021
ZnT8 Deficiency Protects From APAP-Induced Acute Liver Injury by Reducing Oxidative Stress Through Upregulating Hepatic Zinc and Metallothioneins.
Frontiers in pharmacology
Su W,Feng M,Liu Y,Cao R,Liu Y,Tang J,Pan K,Lan R,Mao Z
Sat Aug 21 00:00:00 UTC 2021
Resolving the graft ischemia-reperfusion injury during liver transplantation at the single cell resolution.
Cell death & disease
Wang L,Li J,He S,Liu Y,Chen H,He S,Yin M,Zou D,Chen S,Luo T,Yu X,Wan X,Huang S,Guo Z,He X
Tue Jun 08 00:00:00 UTC 2021
Chlorogenic Acid Ameliorates Damage Induced by Fluorene-9-Bisphenol in Porcine Sertoli Cells.
Frontiers in pharmacology
Zhang S,Sun B,Wang D,Liu Y,Li J,Qi J,Zhang Y,Bai C,Liang S
Tue Jun 29 00:00:00 UTC 2021
Melatonin suppresses ER stress-dependent proapoptotic effects via AMPK in bone mesenchymal stem cells during mitochondrial oxidative damage.
Stem cell research & therapy
Fan C,Feng J,Tang C,Zhang Z,Feng Y,Duan W,Zhai M,Yan Z,Zhu L,Feng L,Zhu H,Luo E
Thu Oct 15 00:00:00 UTC 2020
Efficacy of a wound-dressing biomaterial on prevention of postinflammatory hyperpigmentation after suction blister epidermal grafting in stable vitiligo patients: a controlled assessor-blinded clinical study with in vitro bioactivity investigation.
Archives of dermatological research
Liu Z,Jiang M,Zhao J,Wang Q,Zhang C,Gao M,Gu M,Xiang L
Sun Nov 01 00:00:00 UTC 2020
The critical role of the hippocampal NLRP3 inflammasome in social isolation-induced cognitive impairment in male mice.
Neurobiology of learning and memory
Niu L,Luo SS,Xu Y,Wang Z,Luo D,Yang H,Li W,He J,Zhong XL,Liu ZH,Zeng JY,Cao WY,Wan W
Sun Nov 01 00:00:00 UTC 2020
Anti-tumour activity of zinc ionophore pyrithione in human ovarian cancer cells through inhibition of proliferation and migration and promotion of lysosome-mitochondrial apoptosis.
Artificial cells, nanomedicine, and biotechnology
Chen M,Ding Y,Ke Y,Zeng Y,Liu N,Zhong Y,Hua X,Li Z,Xiong Y,Wu C,Yu H
Tue Dec 01 00:00:00 UTC 2020
Indole alkaloids from leaves of Alstonia scholaris (L.) R. Br. protect against emphysema in mice.
Journal of ethnopharmacology
Zhao YL,Yang ZF,Wu BF,Shang JH,Liu YP,Wang XH,Luo XD
Tue Sep 15 00:00:00 UTC 2020
Synthesis and biological evaluations of N'-substituted methylene-4-(quinoline-4-amino) benzoylhydrazides as potential anti-hepatoma agents.
Bioorganic chemistry
Li B,Zhu F,He F,Huang Q,Liu X,Wu T,Zhao T,Qiu Y,Wu Z,Xue Y,Fang M
Sun Mar 01 00:00:00 UTC 2020
Radiosensitization Effect of AGuIX, a Gadolinium-Based Nanoparticle, in Nonsmall Cell Lung Cancer.
ACS applied materials & interfaces
Du Y,Sun H,Lux F,Xie Y,Du L,Xu C,Zhang H,He N,Wang J,Liu Y,Leduc G,Doussineau T,Ji K,Wang Q,Lin Z,Wang Y,Liu Q,Tillement O
Wed Dec 23 00:00:00 UTC 2020
Systemic MCPIP1 deficiency in mice impairs lipid homeostasis.
Current research in pharmacology and drug discovery
Moody J,Yang C,Sedinkin J,Chang Y
Wed Apr 01 00:00:00 UTC 2020
Myricetin ameliorates bleomycin-induced pulmonary fibrosis in mice by inhibiting TGF-β signaling via targeting HSP90β.
Biochemical pharmacology
Li X,Yu H,Liang L,Bi Z,Wang Y,Gao S,Wang M,Li H,Miao Y,Deng R,Ma L,Luan J,Li S,Liu M,Lin J,Zhou H,Yang C
Sat Aug 01 00:00:00 UTC 2020
SSEA3 and Sialyl Lewis a Glycan Expression Is Controlled by B3GALT5 LTR through Lamin A-NFYA and SIRT1-STAT3 Signaling in Human ES Cells.
Cells
Cai BH,Lee HY,Chou CK,Wu PH,Huang HC,Chao CC,Chung HY,Kannagi R
Fri Jan 10 00:00:00 UTC 2020
Anti-Inflammatory and Antioxidant Effect of Eucommia ulmoides Polysaccharide in Hepatic Ischemia-Reperfusion Injury by Regulating ROS and the TLR-4-NF-κB Pathway.
BioMed research international
Gao W,Feng Z,Zhang S,Wu B,Geng X,Fan G,Duan Y,Li K,Liu K,Peng C
Mon Mar 22 00:00:00 UTC 2021
Epigenetic Inactivation of α-Internexin Accelerates Microtubule Polymerization in Colorectal Cancer.
Cancer research
Li Y,Bai L,Yu H,Cai D,Wang X,Huang B,Peng S,Huang M,Cao G,Kaz AM,Grady WM,Wang J,Luo Y
Tue Dec 01 00:00:00 UTC 2020
Cortistatin ameliorates Ang II-induced proliferation of vascular smooth muscle cells by inhibiting autophagy through SSTR3 and SSTR5.
Life sciences
Wang Y,Zhang X,Chen W,Gao L,Li J,Song T,Chi J,Zhang X,Shi Z,Dong Y,Yin X,Liu Y
Wed Jul 15 00:00:00 UTC 2020
Design, synthesis and biological evaluation of a novel tubulin inhibitor SKLB0565 targeting the colchicine binding site.
Bioorganic chemistry
Hu X,Li L,Zhang Q,Wang Q,Feng Z,Xu Y,Xia Y,Yu L
Wed Apr 01 00:00:00 UTC 2020
A novel microRNA and its pfk target control growth length in the freshwater shrimp Neocaridina heteropoda.
The Journal of experimental biology
Li R,Weng J,Ren L,Wang X,Meng Q,Wang L,Sun J
Wed Jul 08 00:00:00 UTC 2020
Metabolomic profiling reveals similar cytotoxic effects and protective functions of quercetin during deoxynivalenol- and 15-acetyl deoxynivalenol-induced cell apoptosis.
Toxicology in vitro : an international journal published in association with BIBRA
Yang YX,Yu S,Jia BX,Liu N,Wu A
Sat Aug 01 00:00:00 UTC 2020
Identification of Transcriptional Variation in Aortic Remodeling Using a Murine Transverse Aortic Constriction (TAC) Model.
Frontiers in cardiovascular medicine
Zhang X,Feng T,Zeng XI,Liang H,Situ B,Zhang Q,Zhou F,Chen Y,Wang T,Cai D,Lin X,Xiu J,Zheng L
Sat Dec 12 00:00:00 UTC 2020
Activation of CRHR1 contributes to cerebral endothelial barrier impairment via cPLA2 phosphorylation in experimental ischemic stroke.
Cellular signalling
Cao C,Zhou J,Wu X,Qian Y,Hong Y,Mu J,Jin L,Zhu C,Li S
Sat Feb 01 00:00:00 UTC 2020
Anti-Inflammatory Effect of Simonsinol on Lipopolysaccharide Stimulated RAW264.7 Cells through Inactivation of NF-κB Signaling Pathway.
Molecules (Basel, Switzerland)
Li LC,Pan ZH,Ning DS,Fu YX
Thu Aug 06 00:00:00 UTC 2020
Chrysin Inhibits Melanoma Tumor Metastasis via Interfering with the FOXM1/β-Catenin Signaling.
Journal of agricultural and food chemistry
Yufei Z,Yuqi W,Binyue H,Lingchen T,Xi C,Hoffelt D,Fuliang H
Wed Sep 02 00:00:00 UTC 2020
Arfgef1 haploinsufficiency in mice alters neuronal endosome composition and decreases membrane surface postsynaptic GABAA receptors.
Neurobiology of disease
Teoh J,Subramanian N,Pero ME,Bartolini F,Amador A,Kanber A,Williams D,Petri S,Yang M,Allen AS,Beal J,Haut SR,Frankel WN
Sat Feb 01 00:00:00 UTC 2020
Lactobacillus johnsonii L531 Ameliorates Escherichia coli-Induced Cell Damage via Inhibiting NLRP3 Inflammasome Activity and Promoting ATG5/ATG16L1-Mediated Autophagy in Porcine Mammary Epithelial Cells.
Veterinary sciences
Zou YJ,Xu JJ,Wang X,Zhu YH,Wu Q,Wang JF
Fri Aug 14 00:00:00 UTC 2020
Exogenous spermine attenuates rat diabetic cardiomyopathy via suppressing ROS-p53 mediated downregulation of calcium-sensitive receptor.
Redox biology
Wang Y,Chen J,Li S,Zhang X,Guo Z,Hu J,Shao X,Song N,Zhao Y,Li H,Yang G,Xu C,Wei C
Fri May 01 00:00:00 UTC 2020
Inhibition of p38 MAPK regulates epileptic severity by decreasing expression levels of A1R and ENT1.
Molecular medicine reports
Zhou X,Chen Q,Huang H,Zhang J,Wang J,Chen Y,Peng Y,Zhang H,Zeng J,Feng Z,Xu Z
Tue Dec 01 00:00:00 UTC 2020
Doxorubicin-polyglycerol-nanodiamond conjugates disrupt STAT3/IL-6-mediated reciprocal activation loop between glioblastoma cells and astrocytes.
Journal of controlled release : official journal of the Controlled Release Society
Chen Z,Yuan SJ,Li K,Zhang Q,Li TF,An HC,Xu HZ,Yue Y,Han M,Xu YH,Komatsu N,Zhao L,Chen X
Fri Apr 10 00:00:00 UTC 2020
Melatonin regulates mitochondrial dynamics and alleviates neuron damage in prion diseases.
Aging
Zhang X,Zhao D,Wu W,Ali Shah SZ,Lai M,Yang D,Li J,Guan Z,Li W,Gao H,Zhao H,Zhou X,Yang L
Wed Jun 10 00:00:00 UTC 2020
The developmental and experience-dependent expression of IGF-2 in mice visual cortex.
Neuroscience letters
Luo Y,Liu Z,Luo S,Wang X,Tao L
Mon Mar 16 00:00:00 UTC 2020
EPS8 regulates an NLRP3 inflammasome-independent caspase-1 activation pathway in monosodium urate crystal-treated RAW264.7 macrophages.
Biochemical and biophysical research communications
Chuang JP,Kao CY,Lee JC,Ling P,Maa MC,Leu TH
Thu Sep 24 00:00:00 UTC 2020
Monophosphoryl lipid A alleviated radiation-induced testicular injury through TLR4-dependent exosomes.
Journal of cellular and molecular medicine
Liu Z,Cao K,Liao Z,Chen Y,Lei X,Wei Q,Liu C,Sun X,Yang Y,Cai J,Gao F
Wed Apr 01 00:00:00 UTC 2020
Integration of Transcriptomics and Metabolomics Reveals the Antitumor Mechanism Underlying Shikonin in Colon Cancer.
Frontiers in pharmacology
Chen Y,Gao Y,Yi X,Zhang J,Chen Z,Wu Y
Tue Dec 08 00:00:00 UTC 2020
Precise base editing with CC context-specificity using engineered human APOBEC3G-nCas9 fusions.
BMC biology
Liu Z,Chen S,Shan H,Jia Y,Chen M,Song Y,Lai L,Li Z
Mon Aug 31 00:00:00 UTC 2020
Autophagy inducer activates Nrf2-ARE pathway to attenuate aberrant alveolarization in neonatal rats with bronchopulmonary dysplasia.
Life sciences
Zhao X,Shi Y,Zhang D,Tong X,Sun Y,Xue X,Fu J
Wed Jul 01 00:00:00 UTC 2020
Beneficial Effects of Crocin against Depression via Pituitary Adenylate Cyclase-Activating Polypeptide.
BioMed research international
Lu L,Wu D,Wang K,Tang J,Chen G
Fri Apr 16 00:00:00 UTC 2021
RNA-binding protein CUGBP1 controls the differential INSR splicing in molecular subtypes of breast cancer cells and affects cell aggressiveness.
Carcinogenesis
Huang G,Song C,Wang N,Qin T,Sui S,Obr A,Zeng L,Wood TL,Leroith D,Li M,Wu Y
Thu Sep 24 00:00:00 UTC 2020
Simvastatin re-sensitizes hepatocellular carcinoma cells to sorafenib by inhibiting HIF-1α/PPAR-γ/PKM2-mediated glycolysis.
Journal of experimental & clinical cancer research : CR
Feng J,Dai W,Mao Y,Wu L,Li J,Chen K,Yu Q,Kong R,Li S,Zhang J,Ji J,Wu J,Mo W,Xu X,Guo C
Thu Jan 30 00:00:00 UTC 2020
PCNA and GSK3β interact with each other to regulate H1299 lung adenocarcinoma cells apoptosis.
Neoplasma
Liu XH,Tang DE,Dai Y,Gao XJ,Liu LX
Wed Jan 01 00:00:00 UTC 2020
Skp2 modulates proliferation, senescence and tumorigenesis of glioma.
Cancer cell international
Wu J,Su HK,Yu ZH,Xi SY,Guo CC,Hu ZY,Qu Y,Cai HP,Zhao YY,Zhao HF,Chen FR,Huang YF,To ST,Feng BH,Sai K,Chen ZP,Wang J
Wed Apr 13 00:00:00 UTC 2022
Flavagline synthetic derivative induces senescence in glioblastoma cancer cells without being toxic to healthy astrocytes.
Scientific reports
Harmouch E,Seitlinger J,Chaddad H,Ubeaud-Sequier G,Barths J,Saidu S,Désaubry L,Grandemange S,Massfelder T,Fuhrmann G,Fioretti F,Dontenwill M,Benkirane-Jessel N,Idoux-Gillet Y
Thu Aug 13 00:00:00 UTC 2020
Repression of FGF signaling is responsible for Dnmt3b inhibition and impaired de novo DNA methylation during early development of in vitro fertilized embryos.
International journal of biological sciences
Fu W,Yue Y,Miao K,Xi G,Zhang C,Wang W,An L,Tian J
Mon Apr 04 00:00:00 UTC 2022
miR-203 inhibits cell proliferation and ERK pathway in prostate cancer by targeting IRS-1.
BMC cancer
Meng Y,Hu X,Li S,Zeng X,Qiu L,Wei M,Wang Z,Han J
Tue Oct 27 00:00:00 UTC 2020
Nucleus-located PDK1 regulates growth, invasion and migration of breast cancer cells.
Life sciences
Gan D,Yue S,Jiang Y,Zhang D,Shi H,Qian H,Zhou T,Fang W,Yao M,Zuo G,Chen T
Wed Jul 15 00:00:00 UTC 2020
Exogenous H2S Induces Hrd1 S-sulfhydration and Prevents CD36 Translocation via VAMP3 Ubiquitylation in Diabetic Hearts.
Aging and disease
Yu M,Du H,Wang B,Chen J,Lu F,Peng S,Sun Y,Liu N,Sun X,Shiyun D,Zhao Y,Wang Y,Zhao D,Lu F,Zhang W
Wed Apr 01 00:00:00 UTC 2020
Crosstalk between androgen and Wnt/β-catenin leads to changes of wool density in FGF5-knockout sheep.
Cell death & disease
Zhang R,Li Y,Jia K,Xu X,Li Y,Zhao Y,Zhang X,Zhang J,Liu G,Deng S,Lian Z
Fri May 29 00:00:00 UTC 2020
Similar Repair Effects of Human Placenta, Bone Marrow Mesenchymal Stem Cells, and Their Exosomes for Damaged SVOG Ovarian Granulosa Cells.
Stem cells international
Chen S,Wang Y,Liao L,Meng L,Li J,Shi C,Han H,Zheng X,Shen H
Tue Apr 19 00:00:00 UTC 2022
Neuropeptide Y1 receptor antagonist promotes osteoporosis and microdamage repair and enhances osteogenic differentiation of bone marrow stem cells via cAMP/PKA/CREB pathway.
Aging
Xie W,Li F,Han Y,Qin Y,Wang Y,Chi X,Xiao J,Li Z
Thu May 07 00:00:00 UTC 2020
HCK can serve as novel prognostic biomarker and therapeutic target for Breast Cancer patients.
International journal of medical sciences
Zhu X,Zhang Y,Bai Y,Gu X,Chen G,Sun L,Wang Y,Qiao X,Ma Q,Zhu T,Bu J,Xue J,Liu C
Mon Aug 02 00:00:00 UTC 2021
Protective effects of Salidroside on spermatogenesis in streptozotocin induced type-1 diabetic male mice by inhibiting oxidative stress mediated blood-testis barrier damage.
Chemico-biological interactions
Jiang YP,Ye RJ,Yang JM,Liu N,Zhang WJ,Ma L,Sun T,Niu JG,Zheng P,Yu JQ
Sun Jan 05 00:00:00 UTC 2020
PCNP promotes ovarian cancer progression by accelerating β-catenin nuclear accumulation and triggering EMT transition.
Journal of cellular and molecular medicine
Dong P,Fu H,Chen L,Zhang S,Zhang X,Li H,Wu D,Ji X
Wed Jul 01 00:00:00 UTC 2020
Protein target identification of ginsenosides in skeletal muscle tissues: discovery of natural small-molecule activators of muscle-type creatine kinase.
Journal of ginseng research
Chen F,Zhu K,Chen L,Ouyang L,Chen C,Gu L,Jiang Y,Wang Z,Lin Z,Zhang Q,Shao X,Dai J,Zhao Y
Fri May 01 00:00:00 UTC 2020
The ubiquitin-specific protease USP8 directly deubiquitinates SQSTM1/p62 to suppress its autophagic activity.
Autophagy
Peng H,Yang F,Hu Q,Sun J,Peng C,Zhao Y,Huang C
Wed Apr 01 00:00:00 UTC 2020
Ubiquitination of NOTCH2 by DTX3 suppresses the proliferation and migration of human esophageal carcinoma.
Cancer science
Ding XY,Hu HY,Huang KN,Wei RQ,Min J,Qi C,Tang H,Qin X
Sat Feb 01 00:00:00 UTC 2020
Synergistic antifibrotic effects of miR-451 with miR-185 partly by co-targeting EphB2 on hepatic stellate cells.
Cell death & disease
Chen X,Zhang D,Wang Y,Chen K,Zhao L,Xu Y,Jiang H,Wang S
Thu May 28 00:00:00 UTC 2020
Adiponectin protects HL-1 cardiomyocytes against rotenone-induced cytotoxicity through AMPK activation.
Toxicology letters
Li B,Zhang B,Liu N,Wu K,Ma Y,Zuo W,Ning Z,Liu Y,Sun C,Xiao Y,Tu T,Liu Q
Tue Dec 15 00:00:00 UTC 2020
MitoQ attenuates brain damage by polarizing microglia towards the M2 phenotype through inhibition of the NLRP3 inflammasome after ICH.
Pharmacological research
Chen W,Guo C,Huang S,Jia Z,Wang J,Zhong J,Ge H,Yuan J,Chen T,Liu X,Hu R,Yin Y,Feng H
Sun Nov 01 00:00:00 UTC 2020
NCAPG Dynamically Coordinates the Myogenesis of Fetal Bovine Tissue by Adjusting Chromatin Accessibility.
International journal of molecular sciences
Hu X,Xing Y,Fu X,Yang Q,Ren L,Wang Y,Li Q,Li J,Zhang L
Thu Feb 13 00:00:00 UTC 2020
The E3 ubiquitin ligase Smurf2 regulates PARP1 stability to alleviate oxidative stress-induced injury in human umbilical vein endothelial cells.
Journal of cellular and molecular medicine
Qian H,Zhang N,Wu B,Wu S,You S,Zhang Y,Sun Y
Wed Apr 01 00:00:00 UTC 2020
Neural Tube Defects and ZIC4 Hypomethylation in Relation to Polycyclic Aromatic Hydrocarbon Exposure.
Frontiers in cell and developmental biology
Huang Y,Lin S,Wang C,Pi X,Jin L,Li Z,Wang L,Ren A
Mon Apr 18 00:00:00 UTC 2022
Oxidative stress-mediated hepatotoxicity in rats induced by ethanol extracts of different parts of Chloranthus serratus.
Pharmaceutical biology
Sun S,Wang Y,Du Y,Sun Q,He L,Zhu E,Li J
Tue Dec 01 00:00:00 UTC 2020
Mbd2 Mediates Retinal Cell Apoptosis by Targeting the lncRNA Mbd2-AL1/miR-188-3p/Traf3 Axis in Ischemia/Reperfusion Injury.
Molecular therapy. Nucleic acids
Ge Y,Zhang R,Feng Y,Li H
Fri Mar 06 00:00:00 UTC 2020
Leptin mediates protection of hydrogen sulfide against 6-hydroxydopamine-induced Parkinson's disease: Involving enhancement in Warburg effect.
Neurochemistry international
Yang SQ,Tian Q,Li D,He SQ,Hu M,Liu SY,Zou W,Chen YJ,Zhang P,Tang XQ
Fri May 01 00:00:00 UTC 2020
Hypoxia induces tumor cell growth and angiogenesis in non-small cell lung carcinoma via the Akt-PDK1-HIF1α-YKL-40 pathway.
Translational cancer research
Miao Y,Wang W,Dong Y,Hu J,Wei K,Yang S,Lai X,Tang H
Wed Apr 01 00:00:00 UTC 2020
G Protein-Coupled Receptor 109A Maintains the Intestinal Integrity and Protects Against ETEC Mucosal Infection by Promoting IgA Secretion.
Frontiers in immunology
Gong Y,Jin X,Yuan B,Lv Y,Yan G,Liu M,Xie C,Liu J,Tang Y,Gao H,Zhu Y,Huang Y,Wang W
Tue Jun 22 00:00:00 UTC 2021
Lnc-M2 controls M2 macrophage differentiation via the PKA/CREB pathway.
Molecular immunology
Chen Y,Li H,Ding T,Li J,Zhang Y,Wang J,Yang X,Chong T,Long Y,Li X,Gao F,Lyu X
Sat Aug 01 00:00:00 UTC 2020
bta-miR-23a Regulates the Myogenic Differentiation of Fetal Bovine Skeletal Muscle-Derived Progenitor Cells by Targeting MDFIC Gene.
Genes
Hu X,Xing Y,Ren L,Wang Y,Li Q,Yang Q,Du M,Xu L,Willems L,Li J,Zhang L
Tue Oct 20 00:00:00 UTC 2020
Tongmai formula improves cardiac function via regulating mitochondrial quality control in the myocardium with ischemia/reperfusion injury.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhao Y,Guo R,Li L,Li S,Fan G,Zhao X,Wang Y
Tue Dec 01 00:00:00 UTC 2020
WWP2 regulates SIRT1-STAT3 acetylation and phosphorylation involved in hypertensive angiopathy.
Journal of cellular and molecular medicine
Zhang Y,You S,Tian Y,Lu S,Cao L,Sun Y,Zhang N
Sat Aug 01 00:00:00 UTC 2020
A Novel Mechanism of bta-miR-210 in Bovine Early Intramuscular Adipogenesis.
Genes
Ren L,Li Q,Hu X,Yang Q,Du M,Xing Y,Wang Y,Li J,Zhang L
Fri May 29 00:00:00 UTC 2020
Identification of long-term survival-associated gene in breast cancer.
Aging
Ning S,Li H,Qiao K,Wang Q,Shen M,Kang Y,Yin Y,Liu J,Liu L,Hou S,Wang J,Xu S,Pang D
Tue Oct 20 00:00:00 UTC 2020
Pizotifen inhibits the proliferation and invasion of gastric cancer cells.
Experimental and therapeutic medicine
Jiang Y,Wang W,Wu X,Shi J
Sat Feb 01 00:00:00 UTC 2020
Hepatic HuR modulates lipid homeostasis in response to high-fat diet.
Nature communications
Zhang Z,Zong C,Jiang M,Hu H,Cheng X,Ni J,Yi X,Jiang B,Tian F,Chang MW,Su W,Zhu L,Li J,Xiang X,Miao C,Gorospe M,de Cabo R,Dou Y,Ju Z,Yang J,Jiang C,Yang Z,Wang W
Tue Jun 16 00:00:00 UTC 2020
Neddylation promotes protein translocation between the cytoplasm and nucleus.
Biochemical and biophysical research communications
Li S,Fang W,Cui Y,Shi H,Chen J,Li L,Zhang L,Zhang X
Thu Sep 03 00:00:00 UTC 2020
Cyclophilin A plays an important role in embryo implantation through activating Stat3.
Reproduction (Cambridge, England)
Yu T,Lin S,Xu R,Du TX,Li Y,Gao H,Diao HL,Zhang XH
Tue Sep 01 00:00:00 UTC 2020
Control the fate of human umbilical cord mesenchymal stem cells with dual-enzymatically cross-linked gelatin hydrogels for potential applications in nerve regeneration.
Journal of tissue engineering and regenerative medicine
Li J,Gao F,Ma S,Zhang Y,Zhang J,Guan F,Yao M
Tue Sep 01 00:00:00 UTC 2020
Control the fate of human umbilical cord mesenchymal stem cells with dual-enzymatically cross-linked gelatin hydrogels for potential applications in nerve regeneration.
Journal of tissue engineering and regenerative medicine
Li J,Gao F,Ma S,Zhang Y,Zhang J,Guan F,Yao M
Tue Sep 01 00:00:00 UTC 2020
Blood coagulation factor VIII D1241E polymorphism leads to a weak malectin interaction and reduction of factor VIII posttranslational modification and secretion.
Experimental cell research
Jiang N,Xiao Y,Liu Y,Liu W,Liu S
Tue Dec 01 00:00:00 UTC 2020
Sodium butyrate causes α-synuclein degradation by an Atg5-dependent and PI3K/Akt/mTOR-related autophagy pathway.
Experimental cell research
Qiao CM,Sun MF,Jia XB,Shi Y,Zhang BP,Zhou ZL,Zhao LP,Cui C,Shen YQ
Sat Feb 01 00:00:00 UTC 2020
IGFBP6 regulates vascular smooth muscle cell proliferation and morphology via cyclin E-CDK2.
Journal of cellular physiology
Wang Z,Qi Y,Wang R,Wu W,Li Z,Wang M,Liu R,Zhang C,Li W,Wang S
Tue Dec 01 00:00:00 UTC 2020
Regulation of indoleamine 2, 3-dioxygenase in hippocampal microglia by NLRP3 inflammasome in lipopolysaccharide-induced depressive-like behaviors.
The European journal of neuroscience
Zhang S,Zong Y,Ren Z,Hu J,Wu X,Xiao H,Qin S,Zhou G,Ma Y,Zhang Y,Yu J,Wang K,Lu G,Liu Q
Tue Dec 01 00:00:00 UTC 2020
The B7-H4 gene induces immune escape partly via upregulating the PD-1/Stat3 pathway in non-small cell lung cancer.
Human immunology
Yuan L,Ye J,Fan D
Fri May 01 00:00:00 UTC 2020
Circular RNA hsa_circ_0008285 inhibits colorectal cancer cell proliferation and migration via the miR-382-5p/PTEN axis.
Biochemical and biophysical research communications
Wang J,Luo J,Liu G,Li X
Thu Jun 25 00:00:00 UTC 2020
Possible Roles of Proinflammatory Signaling in Keratinocytes Through Aryl Hydrocarbon Receptor Ligands for the Development of Squamous Cell Carcinoma.
Frontiers in immunology
Sato Y,Fujimura T,Hidaka T,Lyu C,Tanita K,Matsushita S,Yamamoto M,Aiba S
Wed Apr 28 00:00:00 UTC 2021
Inhibition of histone acetyltransferase GCN5 extends lifespan in both yeast and human cell lines.
Aging cell
Huang B,Zhong D,Zhu J,An Y,Gao M,Zhu S,Dang W,Wang X,Yang B,Xie Z
Wed Apr 01 00:00:00 UTC 2020
miR-361-5p Mediates SMAD4 to Promote Porcine Granulosa Cell Apoptosis through VEGFA.
Biomolecules
Ma M,Zhang J,Gao X,Yao W,Li Q,Pan Z
Fri Sep 04 00:00:00 UTC 2020
Homocysteine downregulates cardiac homeobox transcription factor NKX2.5 via IGFBP5.
American journal of physiology. Heart and circulatory physiology
Lai G,Wang L,Li Z,Zhao Y
Tue Dec 01 00:00:00 UTC 2020
Overexpression of NRF1-742 or NRF1-772 Reduces Arsenic-Induced Cytotoxicity and Apoptosis in Human HaCaT Keratinocytes.
International journal of molecular sciences
Wang S,Cheng H,Wang L,Zhao R,Guan D
Mon Mar 16 00:00:00 UTC 2020
Laminar Flow Protects Vascular Endothelial Tight Junctions and Barrier Function via Maintaining the Expression of Long Non-coding RNA MALAT1.
Frontiers in bioengineering and biotechnology
Yang F,Zhang Y,Zhu J,Wang J,Jiang Z,Zhao C,Yang Q,Huang Y,Yao W,Pang W,Han L,Zhou J
Mon Sep 28 00:00:00 UTC 2020
Anti-neuroinflammatory effect of 3,4-dihydroxybenzaldehyde in ischemic stroke.
International immunopharmacology
Li X,Xiang B,Shen T,Xiao C,Dai R,He F,Lin Q
Tue Mar 03 00:00:00 UTC 2020
Yueju-Ganmaidazao Decoction confers rapid antidepressant-like effects and the involvement of suppression of NMDA/NO/cGMP signaling.
Journal of ethnopharmacology
Zhang H,Sun Y,Qian S,Ge R,Guo X,Shen Q,Sheng L,Nie C,Zhang Y,Yao Y,Zhou T,Wang W,Xue W,Chen G
Wed Mar 25 00:00:00 UTC 2020
Interplay of m6A and H3K27 trimethylation restrains inflammation during bacterial infection.
Science advances
Wu C,Chen W,He J,Jin S,Liu Y,Yi Y,Gao Z,Yang J,Yang J,Cui J,Zhao W
Sat Aug 01 00:00:00 UTC 2020
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 UTC 2019
KIAA1199 promotes metastasis of colorectal cancer cells via microtubule destabilization regulated by a PP2A/stathmin pathway.
Oncogene
Zhao L,Zhang D,Shen Q,Jin M,Lin Z,Ma H,Huang S,Zhou P,Wu G,Zhang T
Fri Feb 01 00:00:00 UTC 2019
PMP22 Regulates Cholesterol Trafficking and ABCA1-Mediated Cholesterol Efflux.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Zhou Y,Miles JR,Tavori H,Lin M,Khoshbouei H,Borchelt DR,Bazick H,Landreth GE,Lee S,Fazio S,Notterpek L
Wed Jul 03 00:00:00 UTC 2019
Structural and functional damage to the hippocampal neurovascular unit in diabetes-related depression.
Neural regeneration research
Liu J,Wang YH,Li W,Liu L,Yang H,Meng P,Han YS
Fri Feb 01 00:00:00 UTC 2019
Structural and functional damage to the hippocampal neurovascular unit in diabetes-related depression.
Neural regeneration research
Liu J,Wang YH,Li W,Liu L,Yang H,Meng P,Han YS
Fri Feb 01 00:00:00 UTC 2019
S1PR2 antagonist alleviates oxidative stress-enhanced brain endothelial permeability by attenuating p38 and Erk1/2-dependent cPLA2 phosphorylation.
Cellular signalling
Cao C,Dai L,Mu J,Wang X,Hong Y,Zhu C,Jin L,Li S
Tue Jan 01 00:00:00 UTC 2019
S1PR2 antagonist alleviates oxidative stress-enhanced brain endothelial permeability by attenuating p38 and Erk1/2-dependent cPLA2 phosphorylation.
Cellular signalling
Cao C,Dai L,Mu J,Wang X,Hong Y,Zhu C,Jin L,Li S
Tue Jan 01 00:00:00 UTC 2019
LXRα Promotes the Differentiation of Human Gastric Cancer Cells through Inactivation of Wnt/β-catenin Signaling.
Journal of Cancer
Gao Y,Chen Z,Wang R,Tan X,Huang C,Chen G,Chen Z
Wed Sep 30 00:00:00 UTC 2020
LXRα Promotes the Differentiation of Human Gastric Cancer Cells through Inactivation of Wnt/β-catenin Signaling.
Journal of Cancer
Gao Y,Chen Z,Wang R,Tan X,Huang C,Chen G,Chen Z
Wed Sep 30 00:00:00 UTC 2020
Activation of brain endothelium by Escherichia coli K1 virulence factor cglD promotes polymorphonuclear leukocyte transendothelial migration.
Medical microbiology and immunology
Zhang K,Shi MJ,Niu Z,Chen X,Wei JY,Miao ZW,Zhao WD,Chen YH
Fri Feb 01 00:00:00 UTC 2019
Activation of brain endothelium by Escherichia coli K1 virulence factor cglD promotes polymorphonuclear leukocyte transendothelial migration.
Medical microbiology and immunology
Zhang K,Shi MJ,Niu Z,Chen X,Wei JY,Miao ZW,Zhao WD,Chen YH
Fri Feb 01 00:00:00 UTC 2019
Echinacoside protects against MPTP/MPP+-induced neurotoxicity via regulating autophagy pathway mediated by Sirt1.
Metabolic brain disease
Chen C,Xia B,Tang L,Wu W,Tang J,Liang Y,Yang H,Zhang Z,Lu Y,Chen G,Yang Y,Zhao Y
Fri Feb 01 00:00:00 UTC 2019
Echinacoside protects against MPTP/MPP+-induced neurotoxicity via regulating autophagy pathway mediated by Sirt1.
Metabolic brain disease
Chen C,Xia B,Tang L,Wu W,Tang J,Liang Y,Yang H,Zhang Z,Lu Y,Chen G,Yang Y,Zhao Y
Fri Feb 01 00:00:00 UTC 2019
EIF4A2 interacts with the membrane protein of transmissible gastroenteritis coronavirus and plays a role in virus replication.
Research in veterinary science
Song Z,Yang Y,Wang L,Wang K,Ran L,Xie Y,Huang L,Yang Z,Yuan P,Yu Q
Mon Apr 01 00:00:00 UTC 2019
EIF4A2 interacts with the membrane protein of transmissible gastroenteritis coronavirus and plays a role in virus replication.
Research in veterinary science
Song Z,Yang Y,Wang L,Wang K,Ran L,Xie Y,Huang L,Yang Z,Yuan P,Yu Q
Mon Apr 01 00:00:00 UTC 2019
BH3 mimetic ABT-263 enhances the anticancer effects of apigenin in tumor cells with activating EGFR mutation.
Cell & bioscience
Zhan Y,Wang Y,Qi M,Liang P,Ma Y,Li T,Li H,Dai C,An Z,Qi Y,Wu H,Shao H
Sat Apr 09 00:00:00 UTC 2022
Aloperine suppresses human pulmonary vascular smooth muscle cell proliferation via inhibiting inflammatory response.
The Chinese journal of physiology
Chang Z,Zhang P,Zhang M,Jun F,Hu Z,Yang J,Wu Y,Zhou R
Tue Oct 01 00:00:00 UTC 2019
An improved Tet-on system in microRNA overexpression and CRISPR/Cas9-mediated gene editing.
Journal of animal science and biotechnology
Kang K,Huang L,Li Q,Liao X,Dang Q,Yang Y,Luo J,Zeng Y,Li L,Gou D
Fri Apr 08 00:00:00 UTC 2022
A Comparative Peptidomic Characterization of Cultured Skeletal Muscle Tissues Derived From db/db Mice.
Frontiers in endocrinology
Wu Y,Han M,Wang Y,Gao Y,Cui X,Xu P,Ji C,Zhong T,You L,Zeng Y
Thu Nov 21 00:00:00 UTC 2019
RMI1 contributes to DNA repair and to the tolerance to camptothecin.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Fang L,Sun X,Wang Y,Du L,Ji K,Wang J,He N,Liu Y,Wang Q,Zhai H,Hao J,Xu C,Liu Q
Mon Apr 01 00:00:00 UTC 2019
Deregulated lncRNA expression profile in the mouse lung adenocarcinomas with KRAS-G12D mutation and P53 knockout.
Journal of cellular and molecular medicine
Zhang M,Jiang N,Cui R,Du S,Ou H,Chen T,Ge R,Ma D,Zhang J
Tue Oct 01 00:00:00 UTC 2019
Developmental nicotine exposure elicits multigenerational disequilibria in proBDNF proteolysis and glucocorticoid signaling in the frontal cortices, striata, and hippocampi of adolescent mice.
Biochemical pharmacology
Buck JM,O'Neill HC,Stitzel JA
Tue Oct 01 00:00:00 UTC 2019
USP11 promotes growth and metastasis of colorectal cancer via PPP1CA-mediated activation of ERK/MAPK signaling pathway.
EBioMedicine
Sun H,Ou B,Zhao S,Liu X,Song L,Liu X,Wang R,Peng Z
Tue Oct 01 00:00:00 UTC 2019
The CRISPR-Cas13a Gene-Editing System Induces Collateral Cleavage of RNA in Glioma Cells.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wang Q,Liu X,Zhou J,Yang C,Wang G,Tan Y,Wu Y,Zhang S,Yi K,Kang C
Wed Oct 16 00:00:00 UTC 2019
Protective effect of hyperoside against renal ischemia-reperfusion injury via modulating mitochondrial fission, oxidative stress, and apoptosis.
Free radical research
Wu L,Li Q,Liu S,An X,Huang Z,Zhang B,Yuan Y,Xing C
Mon Jul 01 00:00:00 UTC 2019
Effects of Betaine on LPS-Stimulated Activation of Microglial M1/M2 Phenotypes by Suppressing TLR4/NF-κB Pathways in N9 Cells.
Molecules (Basel, Switzerland)
Shi H,Wang XL,Quan HF,Yan L,Pei XY,Wang R,Peng XD
Mon Jan 21 00:00:00 UTC 2019
Neuroprotective Effect of Echinacoside in Subacute Mouse Model of Parkinson's Disease.
BioMed research international
Liang Y,Chen C,Xia B,Wu W,Tang J,Chen Q,Tang L,Yang H,Zhang Z,Lu Y,Yang Y,Zhao Y
Thu Jun 13 00:00:00 UTC 2019
Hyperbaric oxygen therapy reduces apoptosis and dendritic/synaptic degeneration via the BDNF/TrkB signaling pathways in SCI rats.
Life sciences
Ying X,Tu W,Li S,Wu Q,Chen X,Zhou Y,Hu J,Yang G,Jiang S
Mon Jul 15 00:00:00 UTC 2019
Overexpression of ANLN in lung adenocarcinoma is associated with metastasis.
Thoracic cancer
Xu J,Zheng H,Yuan S,Zhou B,Zhao W,Pan Y,Qi D
Thu Aug 01 00:00:00 UTC 2019
Doxorubicin conjugated with nanodiamonds and in free form commit glioblastoma cells to heterodromous fates.
Nanomedicine (London, England)
Chen Z,Wang C,Li TF,Li K,Yue Y,Liu X,Xu HZ,Wen Y,Zhang Q,Han M,Komatsu N,Xu YH,Zhao L,Chen X
Fri Feb 01 00:00:00 UTC 2019
Cardioprotective effects of galectin-3 inhibition against ischemia/reperfusion injury.
European journal of pharmacology
Mo D,Tian W,Zhang HN,Feng YD,Sun Y,Quan W,Hao XW,Wang XY,Liu XX,Li C,Cao W,Liu WJ,Li XQ
Fri Nov 15 00:00:00 UTC 2019
Interactions between β-cyclodextrin and tea catechins, and potential anti-osteoclastogenesis activity of the (-)-epigallocatechin-3-gallate-β-cyclodextrin complex.
RSC advances
Xu H,Liu T,Xu J,Li J,Chen F,Xiang Z,Huang Y,Zhang D,Hu L,Zhang B,Zi C,Wang X,Sheng J
Tue Sep 03 00:00:00 UTC 2019
Casp8 hypomethylation and neural tube defects in association with polycyclic aromatic hydrocarbon exposure.
Clinical epigenetics
Huang Y,Ren A,Wang L,Jin L,Lin S,Li Z,McDonald JA
Tue May 07 00:00:00 UTC 2019
Leonurine promotes neurite outgrowth and neurotrophic activity by modulating the GR/SGK1 signaling pathway in cultured PC12 cells.
Neuroreport
Meng P,Zhu Q,Yang H,Liu D,Lin X,Liu J,Fan J,Liu X,Su W,Liu L,Wang Y,Cai X
Wed Mar 06 00:00:00 UTC 2019
Resveratrol protects against isoproterenol induced myocardial infarction in rats through VEGF-B/AMPK/eNOS/NO signalling pathway.
Free radical research
Feng L,Ren J,Li Y,Yang G,Kang L,Zhang S,Ma C,Li J,Liu J,Yang L,Qi Z
Tue Jan 01 00:00:00 UTC 2019
The protective effects of XH-105 against radiation-induced intestinal injury.
Journal of cellular and molecular medicine
Cheng Y,Dong Y,Hou Q,Wu J,Zhang W,Tian H,Li D
Fri Mar 01 00:00:00 UTC 2019
Combinatory FK506 and Minocycline Treatment Alleviates Prion-Induced Neurodegenerative Events via Caspase-Mediated MAPK-NRF2 Pathway.
International journal of molecular sciences
Shah SZA,Zhao D,Taglialatela G,Hussain T,Dong H,Sabir N,Mangi MH,Wu W,Lai M,Zhang X,Duan Y,Wang L,Zhou X,Yang L
Wed Mar 06 00:00:00 UTC 2019
Inhibition of dendritic spine extension through acrolein conjugation with α-, β-tubulin proteins.
The international journal of biochemistry & cell biology
Uemura T,Suzuki T,Ko K,Watanabe K,Dohmae N,Sakamoto A,Terui Y,Toida T,Kashiwagi K,Igarashi K
Thu Aug 01 00:00:00 UTC 2019
Inhibition of dendritic spine extension through acrolein conjugation with α-, β-tubulin proteins.
The international journal of biochemistry & cell biology
Uemura T,Suzuki T,Ko K,Watanabe K,Dohmae N,Sakamoto A,Terui Y,Toida T,Kashiwagi K,Igarashi K
Thu Aug 01 00:00:00 UTC 2019
Inhibition of dendritic spine extension through acrolein conjugation with α-, β-tubulin proteins.
The international journal of biochemistry & cell biology
Uemura T,Suzuki T,Ko K,Watanabe K,Dohmae N,Sakamoto A,Terui Y,Toida T,Kashiwagi K,Igarashi K
Thu Aug 01 00:00:00 UTC 2019
(-)-Epigallocatechin-3-gallate derivatives combined with cisplatin exhibit synergistic inhibitory effects on non-small-cell lung cancer cells.
Cancer cell international
Wang J,Sun P,Wang Q,Zhang P,Wang Y,Zi C,Wang X,Sheng J
Sun Apr 10 00:00:00 UTC 2022
Lipoxin A4 Regulates Lipopolysaccharide-Induced BV2 Microglial Activation and Differentiation via the Notch Signaling Pathway.
Frontiers in cellular neuroscience
Wu J,Ding DH,Li QQ,Wang XY,Sun YY,Li LJ
Tue Sep 29 00:00:00 UTC 2020
Sevoflurane inhibits the progression of ovarian cancer through down-regulating stanniocalcin 1 (STC1).
Cancer cell international
Zhang C,Wang B,Wang X,Sheng X,Cui Y
Tue Apr 12 00:00:00 UTC 2022
Exosomal transfer of miR-501 confers doxorubicin resistance and tumorigenesis via targeting of BLID in gastric cancer.
Cancer letters
Liu X,Lu Y,Xu Y,Hou S,Huang J,Wang B,Zhao J,Xia S,Fan S,Yu X,Du Y,Hou L,Li Z,Ding Z,An S,Huang B,Li L,Tang J,Ju J,Guan H,Song B
Tue Sep 10 00:00:00 UTC 2019
Integrating yeast chemical genomics and mammalian cell pathway analysis.
Acta pharmacologica Sinica
Zhou FL,Li SC,Zhu Y,Guo WJ,Shao LJ,Nelson J,Simpkins S,Yang DH,Liu Q,Yashiroda Y,Xu JB,Fan YY,Yue JM,Yoshida M,Xia T,Myers CL,Boone C,Wang MW
Sun Sep 01 00:00:00 UTC 2019
Exosomes from human adipose-derived stem cells promote sciatic nerve regeneration via optimizing Schwann cell function.
Journal of cellular physiology
Chen J,Ren S,Duscher D,Kang Y,Liu Y,Wang C,Yuan M,Guo G,Xiong H,Zhan P,Wang Y,Machens HG,Chen Z
Sun Dec 01 00:00:00 UTC 2019
Sirt2 epigenetically down-regulates PDGFRα expression and promotes CG4 cell differentiation.
Cell cycle (Georgetown, Tex.)
Fang N,Cheng J,Zhang C,Chen K,Zhang C,Hu Z,Bi R,Furber KL,Thangaraj M,Nazarali AJ,Ji S
Wed May 01 00:00:00 UTC 2019
Role of Nkx2.5 in H2O2-induced Nsd1 suppression.
Cell stress & chaperones
Liang X,Chu G,Wang L,Lai G,Zhao Y
Mon Jul 01 00:00:00 UTC 2019
Transketolase (TKT) activity and nuclear localization promote hepatocellular carcinoma in a metabolic and a non-metabolic manner.
Journal of experimental & clinical cancer research : CR
Qin Z,Xiang C,Zhong F,Liu Y,Dong Q,Li K,Shi W,Ding C,Qin L,He F
Thu Apr 11 00:00:00 UTC 2019
Water-separated part of Chloranthus serratus alleviates lipopolysaccharide- induced RAW264.7 cell injury mainly by regulating the MAPK and Nrf2/HO-1 inflammatory pathways.
BMC complementary and alternative medicine
Sun S,Du Y,Yin C,Suo X,Wang R,Xia R,Zhang X
Mon Dec 02 00:00:00 UTC 2019
Sulforaphane Inhibits the Acquisition of Tobacco Smoke-Induced Lung Cancer Stem Cell-Like Properties via the IL-6/ΔNp63α/Notch Axis.
Theranostics
Xie C,Zhu J,Jiang Y,Chen J,Wang X,Geng S,Wu J,Zhong C,Li X,Meng Z
Tue Aug 25 00:00:00 UTC 2020
Fuplatin: An Efficient and Low-Toxic Dual-Prodrug.
Journal of medicinal chemistry
Zhang R,Song XQ,Liu RP,Ma ZY,Xu JY
Thu May 09 00:00:00 UTC 2019
Long noncoding RNA CCAT2 promotes hepatocellular carcinoma proliferation and metastasis through up-regulation of NDRG1.
Experimental cell research
Liu Y,Wang D,Li Y,Yan S,Dang H,Yue H,Ling J,Chen F,Zhao Y,Gou L,Tang P,Huang A,Tang H
Sat Jun 01 00:00:00 UTC 2019
An LTR retrotransposon-derived lncRNA interacts with RNF169 to promote homologous recombination.
EMBO reports
Deng B,Xu W,Wang Z,Liu C,Lin P,Li B,Huang Q,Yang J,Zhou H,Qu L
Tue Nov 05 00:00:00 UTC 2019
Inhibitory Effects of Oxymatrine on Transdifferentiation of Neonatal Rat Cardiac Fibroblasts to Myofibroblasts Induced by Aldosterone via Keap1/Nrf2 Signaling Pathways In Vitro.
Medical science monitor : international medical journal of experimental and clinical research
Yang Y,Chen S,Tao L,Gan S,Luo H,Xu Y,Shen X
Sat Jul 20 00:00:00 UTC 2019
YAP mediates the positive regulation of hnRNPK on the lung adenocarcinoma H1299 cell growth.
Acta biochimica et biophysica Sinica
Xu L,Zhang T,Huang W,Liu X,Lu J,Gao X,Zhang YF,Liu L
Wed Jul 10 00:00:00 UTC 2019
Matrix Metalloproteases-Mediated Cleavage on β-Dystroglycan May Play a Key Role in the Blood-Brain Barrier After Intracerebral Hemorrhage in Rats.
Medical science monitor : international medical journal of experimental and clinical research
Zhang X,Gu Y,Li P,Jiang A,Sheng X,Jin X,Shi Y,Li G
Mon Jan 28 00:00:00 UTC 2019
Astaxanthin inhibits proliferation and induces apoptosis of LX‑2 cells by regulating the miR‑29b/Bcl‑2 pathway.
Molecular medicine reports
Zhu S,Wang T,Luo F,Li H,Jia Q,He T,Wu H,Zou T
Wed May 01 00:00:00 UTC 2019
Oxidation derivative of (-)-epigallocatechin-3-gallate (EGCG) inhibits RANKL-induced osteoclastogenesis by suppressing RANK signaling pathways in RAW 264.7 cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Xu H,Liu T,Li J,Xu J,Chen F,Hu L,Zhang B,Zi C,Wang X,Sheng J
Tue Oct 01 00:00:00 UTC 2019
The Capsid Protein VP1 of Coxsackievirus B Induces Cell Cycle Arrest by Up-Regulating Heat Shock Protein 70.
Frontiers in microbiology
Wang Y,Zhao S,Chen Y,Wang T,Dong C,Wo X,Zhang J,Dong Y,Xu W,Feng X,Qu C,Wang Y,Zhong Z,Zhao W
Thu Oct 01 00:00:00 UTC 2020
Catalpol prevents denervated muscular atrophy related to the inhibition of autophagy and reduces BAX/BCL2 ratio via mTOR pathway.
Drug design, development and therapy
Wang Y,Shao Y,Gao Y,Wan G,Wan D,Zhu H,Qiu Y,Ye X
Wed May 29 00:00:00 UTC 2019
Catalpol prevents denervated muscular atrophy related to the inhibition of autophagy and reduces BAX/BCL2 ratio via mTOR pathway.
Drug design, development and therapy
Wang Y,Shao Y,Gao Y,Wan G,Wan D,Zhu H,Qiu Y,Ye X
Wed May 29 00:00:00 UTC 2019
TLR4/NF-κB Signaling Induces GSDMD-Related Pyroptosis in Tubular Cells in Diabetic Kidney Disease.
Frontiers in endocrinology
Wang Y,Zhu X,Yuan S,Wen S,Liu X,Wang C,Qu Z,Li J,Liu H,Sun L,Liu F
Wed Oct 23 00:00:00 UTC 2019
The Binding of PD-L1 and Akt Facilitates Glioma Cell Invasion Upon Starvation via Akt/Autophagy/F-Actin Signaling.
Frontiers in oncology
Chen RQ,Xu XH,Liu F,Li CY,Li YJ,Li XR,Jiang GY,Hu F,Liu D,Pan F,Qiu XY,Chen XQ
Thu Oct 01 00:00:00 UTC 2020
Anti-TGEV Miller Strain Infection Effect of Lactobacillus plantarum Supernatant Based on the JAK-STAT1 Signaling Pathway.
Frontiers in microbiology
Wang K,Ran L,Yan T,Niu Z,Kan Z,Zhang Y,Yang Y,Xie L,Huang S,Yu Q,Wu D,Song Z
Thu Oct 01 00:00:00 UTC 2020
HIF-1α contributes to Ang II-induced inflammatory cytokine production in podocytes.
BMC pharmacology & toxicology
Huang H,Fan Y,Gao Z,Wang W,Shao N,Zhang L,Yang Y,Zhu W,Chen Z,Hu J,Ding G
Thu Oct 17 00:00:00 UTC 2019
The oncogenic role of CB2 in the progression of non-small-cell lung cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Xu S,Ma H,Bo Y,Shao M
Sun Sep 01 00:00:00 UTC 2019
Protein kinase C-iota-mediated glycolysis promotes non-small-cell lung cancer progression.
OncoTargets and therapy
Liu L,Lei B,Wang L,Chang C,Yang H,Liu J,Huang G,Xie W
Sat Apr 09 00:00:00 UTC 2022
Long-Term Effects of Zinc Deficiency and Zinc Supplementation on Developmental Seizure-Induced Brain Damage and the Underlying GPR39/ZnT-3 and MBP Expression in the Hippocampus.
Frontiers in neuroscience
Chen NN,Zhao DJ,Sun YX,Wang DD,Ni H
Wed Sep 30 00:00:00 UTC 2020
Hypoxic postconditioning activates the Wnt/β-catenin pathway and protects against transient global cerebral ischemia through Dkk1 Inhibition and GSK-3β inactivation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhan L,Liu D,Wen H,Hu J,Pang T,Sun W,Xu E
Thu Aug 01 00:00:00 UTC 2019
Long-term pulmonary exposure to multi-walled carbon nanotubes promotes breast cancer metastatic cascades.
Nature nanotechnology
Lu X,Zhu Y,Bai R,Wu Z,Qian W,Yang L,Cai R,Yan H,Li T,Pandey V,Liu Y,Lobie PE,Chen C,Zhu T
Mon Jul 01 00:00:00 UTC 2019
Involvement of the microglial NLRP3 inflammasome in the anti-inflammatory effect of the antidepressant clomipramine.
Journal of affective disorders
Gong W,Zhang S,Zong Y,Halim M,Ren Z,Wang Y,Ma Y,Li B,Ma L,Zhou G,Yu J,Zhang J,Liu Q
Mon Jul 01 00:00:00 UTC 2019
Crocin induces anti-ischemia in middle cerebral artery occlusion rats and inhibits autophagy by regulating the mammalian target of rapamycin.
European journal of pharmacology
Huang Z,Xu J,Huang X,Sun G,Jiang R,Wu H,Shan X,Bao K,Wu Q,Wu H,Tao W
Thu Aug 15 00:00:00 UTC 2019
Sorafenib kills liver cancer cells by disrupting SCD1-mediated synthesis of monounsaturated fatty acids via the ATP-AMPK-mTOR-SREBP1 signaling pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liu G,Kuang S,Cao R,Wang J,Peng Q,Sun C
Sun Sep 01 00:00:00 UTC 2019
Protective effects of betaine on diabetic induced disruption of the male mice blood-testis barrier by regulating oxidative stress-mediated p38 MAPK pathways.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Jiang YP,Yang JM,Ye RJ,Liu N,Zhang WJ,Ma L,Zheng P,Niu JG,Liu P,Yu JQ
Sun Dec 01 00:00:00 UTC 2019
PTEN-L promotes type I interferon responses and antiviral immunity.
Cellular & molecular immunology
Cao Y,Wang H,Yang L,Zhang Z,Li C,Yuan X,Bu L,Chen L,Chen Y,Li CM,Guo D
Mon Jan 01 00:00:00 UTC 2018
PTEN-L promotes type I interferon responses and antiviral immunity.
Cellular & molecular immunology
Cao Y,Wang H,Yang L,Zhang Z,Li C,Yuan X,Bu L,Chen L,Chen Y,Li CM,Guo D
Mon Jan 01 00:00:00 UTC 2018
KPNB1 inhibition disrupts proteostasis and triggers unfolded protein response-mediated apoptosis in glioblastoma cells.
Oncogene
Zhu ZC,Liu JW,Li K,Zheng J,Xiong ZQ
Tue May 01 00:00:00 UTC 2018
KPNB1 inhibition disrupts proteostasis and triggers unfolded protein response-mediated apoptosis in glioblastoma cells.
Oncogene
Zhu ZC,Liu JW,Li K,Zheng J,Xiong ZQ
Tue May 01 00:00:00 UTC 2018
Theabrownin suppresses in vitro osteoclastogenesis and prevents bone loss in ovariectomized rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu T,Xiang Z,Chen F,Yin D,Huang Y,Xu J,Hu L,Xu H,Wang X,Sheng J
Mon Oct 01 00:00:00 UTC 2018
Theabrownin suppresses in vitro osteoclastogenesis and prevents bone loss in ovariectomized rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu T,Xiang Z,Chen F,Yin D,Huang Y,Xu J,Hu L,Xu H,Wang X,Sheng J
Mon Oct 01 00:00:00 UTC 2018
The Protective Effect of New Compound XH-103 on Radiation-Induced GI Syndrome.
Oxidative medicine and cellular longevity
Dong Y,Cheng Y,Hou Q,Wu J,Li D,Tian H
Wed Dec 19 00:00:00 UTC 2018
The Protective Effect of New Compound XH-103 on Radiation-Induced GI Syndrome.
Oxidative medicine and cellular longevity
Dong Y,Cheng Y,Hou Q,Wu J,Li D,Tian H
Wed Dec 19 00:00:00 UTC 2018
Rapid induction of apoptosis in tumor cells treated with a new platinum(II) complex based on amino-thiazolidinone.
European journal of medicinal chemistry
Song XQ,Liu YH,Shao J,Zhang ZL,Xie CZ,Qiao X,Bao WG,Xu JY
Wed Sep 05 00:00:00 UTC 2018
Rapid induction of apoptosis in tumor cells treated with a new platinum(II) complex based on amino-thiazolidinone.
European journal of medicinal chemistry
Song XQ,Liu YH,Shao J,Zhang ZL,Xie CZ,Qiao X,Bao WG,Xu JY
Wed Sep 05 00:00:00 UTC 2018
Combined treatment with Dendrobium candidum and black tea extract promotes osteoprotective activity in ovariectomized estrogen deficient rats and osteoclast formation.
Life sciences
Wang MY,Shen C,An MF,Xie CQ,Wu X,Zhu QQ,Sun B,Huang YP,Zhao YL,Wang XJ,Sheng J
Tue May 01 00:00:00 UTC 2018
Combined treatment with Dendrobium candidum and black tea extract promotes osteoprotective activity in ovariectomized estrogen deficient rats and osteoclast formation.
Life sciences
Wang MY,Shen C,An MF,Xie CQ,Wu X,Zhu QQ,Sun B,Huang YP,Zhao YL,Wang XJ,Sheng J
Tue May 01 00:00:00 UTC 2018
Apolipoprotein E deficiency accelerates atherosclerosis development in miniature pigs.
Disease models & mechanisms
Fang B,Ren X,Wang Y,Li Z,Zhao L,Zhang M,Li C,Zhang Z,Chen L,Li X,Liu J,Xiong Q,Zhang L,Jin Y,Liu X,Li L,Wei H,Yang H,Li R,Dai Y
Wed Oct 10 00:00:00 UTC 2018
Apolipoprotein E deficiency accelerates atherosclerosis development in miniature pigs.
Disease models & mechanisms
Fang B,Ren X,Wang Y,Li Z,Zhao L,Zhang M,Li C,Zhang Z,Chen L,Li X,Liu J,Xiong Q,Zhang L,Jin Y,Liu X,Li L,Wei H,Yang H,Li R,Dai Y
Wed Oct 10 00:00:00 UTC 2018
CCAAT/enhancer-binding protein β overexpression alleviates myocardial remodelling by regulating angiotensin-converting enzyme-2 expression in diabetes.
Journal of cellular and molecular medicine
Tie Y,Zhai C,Zhang Y,Qin X,Yu F,Li H,Shan M,Zhang C
Thu Mar 01 00:00:00 UTC 2018
CCAAT/enhancer-binding protein β overexpression alleviates myocardial remodelling by regulating angiotensin-converting enzyme-2 expression in diabetes.
Journal of cellular and molecular medicine
Tie Y,Zhai C,Zhang Y,Qin X,Yu F,Li H,Shan M,Zhang C
Thu Mar 01 00:00:00 UTC 2018
Inhibition of Cdk5 rejuvenates inhibitory circuits and restores experience-dependent plasticity in adult visual cortex.
Neuropharmacology
Li Y,Wang L,Zhang X,Huang M,Li S,Wang X,Chen L,Jiang B,Yang Y
Mon Jan 01 00:00:00 UTC 2018
Inhibition of Cdk5 rejuvenates inhibitory circuits and restores experience-dependent plasticity in adult visual cortex.
Neuropharmacology
Li Y,Wang L,Zhang X,Huang M,Li S,Wang X,Chen L,Jiang B,Yang Y
Mon Jan 01 00:00:00 UTC 2018
The antidepressant effects of α-tocopherol are related to activation of autophagy via the AMPK/mTOR pathway.
European journal of pharmacology
Huang X,Wu H,Jiang R,Sun G,Shen J,Ma M,Ma C,Zhang S,Huang Z,Wu Q,Chen G,Tao W
Wed Aug 15 00:00:00 UTC 2018
The antidepressant effects of α-tocopherol are related to activation of autophagy via the AMPK/mTOR pathway.
European journal of pharmacology
Huang X,Wu H,Jiang R,Sun G,Shen J,Ma M,Ma C,Zhang S,Huang Z,Wu Q,Chen G,Tao W
Wed Aug 15 00:00:00 UTC 2018
Fungal β-Glucan Activates the NLRP3 Inflammasome in Human Bronchial Epithelial Cells Through ROS Production.
Inflammation
Huang Y,Hua M,Cui X
Thu Feb 01 00:00:00 UTC 2018
Fungal β-Glucan Activates the NLRP3 Inflammasome in Human Bronchial Epithelial Cells Through ROS Production.
Inflammation
Huang Y,Hua M,Cui X
Thu Feb 01 00:00:00 UTC 2018
NKX2-2 Suppresses Osteosarcoma Metastasis and Proliferation by Downregulating Multiple Target Genes.
Journal of Cancer
Chen H,Liu W,Zhong L,Liao D,Zhang R,Kang T,Wu Y
Sat Apr 09 00:00:00 UTC 2022
NKX2-2 Suppresses Osteosarcoma Metastasis and Proliferation by Downregulating Multiple Target Genes.
Journal of Cancer
Chen H,Liu W,Zhong L,Liao D,Zhang R,Kang T,Wu Y
Sat Apr 09 00:00:00 UTC 2022
Neuroprotective Effect of Anethole Against Neuropathic Pain Induced by Chronic Constriction Injury of the Sciatic Nerve in Mice.
Neurochemical research
Wang B,Zhang G,Yang M,Liu N,Li YX,Ma H,Ma L,Sun T,Tan H,Yu J
Sat Dec 01 00:00:00 UTC 2018
Neuroprotective Effect of Anethole Against Neuropathic Pain Induced by Chronic Constriction Injury of the Sciatic Nerve in Mice.
Neurochemical research
Wang B,Zhang G,Yang M,Liu N,Li YX,Ma H,Ma L,Sun T,Tan H,Yu J
Sat Dec 01 00:00:00 UTC 2018
Horizontal transfer of exosomal CXCR4 promotes murine hepatocarcinoma cell migration, invasion and lymphangiogenesis.
Gene
Li M,Lu Y,Xu Y,Wang J,Zhang C,Du Y,Wang L,Li L,Wang B,Shen J,Tang J,Song B
Thu Nov 15 00:00:00 UTC 2018
Horizontal transfer of exosomal CXCR4 promotes murine hepatocarcinoma cell migration, invasion and lymphangiogenesis.
Gene
Li M,Lu Y,Xu Y,Wang J,Zhang C,Du Y,Wang L,Li L,Wang B,Shen J,Tang J,Song B
Thu Nov 15 00:00:00 UTC 2018
PMN inhibits colorectal cancer cells through inducing mitotic arrest and p53-dependent apoptosis via the inhibition of tubulin polymerization.
Biochemical and biophysical research communications
Wu X,Shen Y
Wed May 23 00:00:00 UTC 2018
PMN inhibits colorectal cancer cells through inducing mitotic arrest and p53-dependent apoptosis via the inhibition of tubulin polymerization.
Biochemical and biophysical research communications
Wu X,Shen Y
Wed May 23 00:00:00 UTC 2018
iTRAQ-Based Proteomics Analysis Reveals the Effect of Rhubarb in Rats with Ischemic Stroke.
BioMed research international
Lin X,Liu T,Li P,He Z,Zhong Y,Cui H,Luo J,Wang Y,Tang T
Tue Dec 11 00:00:00 UTC 2018
iTRAQ-Based Proteomics Analysis Reveals the Effect of Rhubarb in Rats with Ischemic Stroke.
BioMed research international
Lin X,Liu T,Li P,He Z,Zhong Y,Cui H,Luo J,Wang Y,Tang T
Tue Dec 11 00:00:00 UTC 2018
Isoliquiritigenin inhibits cell proliferation and migration through the PI3K/AKT signaling pathway in A549 lung cancer cells.
Oncology letters
Tian T,Sun J,Wang J,Liu Y,Liu H
Thu Nov 01 00:00:00 UTC 2018
Isoliquiritigenin inhibits cell proliferation and migration through the PI3K/AKT signaling pathway in A549 lung cancer cells.
Oncology letters
Tian T,Sun J,Wang J,Liu Y,Liu H
Thu Nov 01 00:00:00 UTC 2018
PAX2 may induce ADAM10 expression in renal tubular epithelial cells and contribute to epithelial-to-mesenchymal transition.
International urology and nephrology
Hou L,Du Y,Zhao C,Wu Y
Sat Sep 01 00:00:00 UTC 2018
PAX2 may induce ADAM10 expression in renal tubular epithelial cells and contribute to epithelial-to-mesenchymal transition.
International urology and nephrology
Hou L,Du Y,Zhao C,Wu Y
Sat Sep 01 00:00:00 UTC 2018
Effect of decabrominated diphenyl ether exposure on spatial learning and memory, the expression and phosphorylation of hippocampal glutamate receptor subunits in adult Sprague-Dawley rats.
The Journal of toxicological sciences
Xiong L,Liyue H,Fancai Z,Maoting L,Ya L,Ting H,Zhen Y,Shanshan Z,Wenwen G,Yan T
Mon Nov 26 00:00:00 UTC 2018
Effect of decabrominated diphenyl ether exposure on spatial learning and memory, the expression and phosphorylation of hippocampal glutamate receptor subunits in adult Sprague-Dawley rats.
The Journal of toxicological sciences
Xiong L,Liyue H,Fancai Z,Maoting L,Ya L,Ting H,Zhen Y,Shanshan Z,Wenwen G,Yan T
Mon Nov 26 00:00:00 UTC 2018
Temporal Proteomic Analysis of Pancreatic β-Cells in Response to Lipotoxicity and Glucolipotoxicity.
Molecular & cellular proteomics : MCP
Li Z,Liu H,Niu Z,Zhong W,Xue M,Wang J,Yang F,Zhou Y,Zhou Y,Xu T,Hou J
Thu Nov 01 00:00:00 UTC 2018
Temporal Proteomic Analysis of Pancreatic β-Cells in Response to Lipotoxicity and Glucolipotoxicity.
Molecular & cellular proteomics : MCP
Li Z,Liu H,Niu Z,Zhong W,Xue M,Wang J,Yang F,Zhou Y,Zhou Y,Xu T,Hou J
Thu Nov 01 00:00:00 UTC 2018
Salvianolic Acid B-Alleviated Angiotensin II Induces Cardiac Fibrosis by Suppressing NF-κB Pathway In Vitro.
Medical science monitor : international medical journal of experimental and clinical research
Wang C,Luo H,Xu Y,Tao L,Chang C,Shen X
Fri Oct 26 00:00:00 UTC 2018
Salvianolic Acid B-Alleviated Angiotensin II Induces Cardiac Fibrosis by Suppressing NF-κB Pathway In Vitro.
Medical science monitor : international medical journal of experimental and clinical research
Wang C,Luo H,Xu Y,Tao L,Chang C,Shen X
Fri Oct 26 00:00:00 UTC 2018
EGFR as a Negative Regulatory Protein Adjusts the Activity and Mobility of NHE3 in the Cell Membrane of IPEC-J2 Cells With TGEV Infection.
Frontiers in microbiology
Yang Z,Ran L,Yuan P,Yang Y,Wang K,Xie L,Huang S,Liu J,Song Z
Wed Sep 30 00:00:00 UTC 2020
Synergistic neuroprotective effect of rasagiline and idebenone against retinal ischemia-reperfusion injury via the Lin28-let-7-Dicer pathway.
Oncotarget
Lei D,Shao Z,Zhou X,Yuan H
Fri Feb 23 00:00:00 UTC 2018
Synergistic neuroprotective effect of rasagiline and idebenone against retinal ischemia-reperfusion injury via the Lin28-let-7-Dicer pathway.
Oncotarget
Lei D,Shao Z,Zhou X,Yuan H
Fri Feb 23 00:00:00 UTC 2018
Effect of Black Tea Extract and Thearubigins on Osteoporosis in Rats and Osteoclast Formation in vitro.
Frontiers in physiology
Liang Q,Lv M,Zhang X,Hu J,Wu Y,Huang Y,Wang X,Sheng J
Mon Sep 28 00:00:00 UTC 2020
Effect of Black Tea Extract and Thearubigins on Osteoporosis in Rats and Osteoclast Formation in vitro.
Frontiers in physiology
Liang Q,Lv M,Zhang X,Hu J,Wu Y,Huang Y,Wang X,Sheng J
Mon Sep 28 00:00:00 UTC 2020
The ubiquitin-specific protease USP8 deubiquitinates and stabilizes Cx43.
The Journal of biological chemistry
Sun J,Hu Q,Peng H,Peng C,Zhou L,Lu J,Huang C
Fri May 25 00:00:00 UTC 2018
The ubiquitin-specific protease USP8 deubiquitinates and stabilizes Cx43.
The Journal of biological chemistry
Sun J,Hu Q,Peng H,Peng C,Zhou L,Lu J,Huang C
Fri May 25 00:00:00 UTC 2018
Phosphorylation of IRS4 by CK1γ2 promotes its degradation by CHIP through the ubiquitin/lysosome pathway.
Theranostics
Li X,Zhong L,Wang Z,Chen H,Liao D,Zhang R,Zhang H,Kang T
Mon Aug 19 00:00:00 UTC 2019
Phosphorylation of IRS4 by CK1γ2 promotes its degradation by CHIP through the ubiquitin/lysosome pathway.
Theranostics
Li X,Zhong L,Wang Z,Chen H,Liao D,Zhang R,Zhang H,Kang T
Mon Aug 19 00:00:00 UTC 2019
Caffeine Promotes Conversion of Palmitic Acid to Palmitoleic Acid by Inducing Expression of fat-5 in Caenorhabditis elegans and scd1 in Mice.
Frontiers in pharmacology
Du X,Huang Q,Guan Y,Lv M,He X,Fang C,Wang X,Sheng J
Thu Oct 01 00:00:00 UTC 2020
Caffeine Promotes Conversion of Palmitic Acid to Palmitoleic Acid by Inducing Expression of fat-5 in Caenorhabditis elegans and scd1 in Mice.
Frontiers in pharmacology
Du X,Huang Q,Guan Y,Lv M,He X,Fang C,Wang X,Sheng J
Thu Oct 01 00:00:00 UTC 2020
hnRNPK modulates selective quality-control autophagy by downregulating the expression of HDAC6 in 293 cells.
International journal of oncology
Li Z,Liu X,Ma J,Zhang T,Gao X,Liu L
Thu Nov 01 00:00:00 UTC 2018
Met is involved in TIGAR-regulated metastasis of non-small-cell lung cancer.
Molecular cancer
Shen M,Zhao X,Zhao L,Shi L,An S,Huang G,Liu J
Sat May 12 00:00:00 UTC 2018
Met is involved in TIGAR-regulated metastasis of non-small-cell lung cancer.
Molecular cancer
Shen M,Zhao X,Zhao L,Shi L,An S,Huang G,Liu J
Sat May 12 00:00:00 UTC 2018
Involvement of NMDA-AKT-mTOR Signaling in Rapid Antidepressant-Like Activity of Chaihu-jia-Longgu-Muli-tang on Olfactory Bulbectomized Mice.
Frontiers in pharmacology
Wang X,Zou Z,Shen Q,Huang Z,Chen J,Tang J,Xue W,Tao W,Wu H,Wang D,Chen G
Thu Oct 01 00:00:00 UTC 2020
Alginate enhances Toll-like receptor 4-mediated phagocytosis by murine RAW264.7 macrophages.
International journal of biological macromolecules
Bi D,Zhou R,Cai N,Lai Q,Han Q,Peng Y,Jiang Z,Tang Z,Lu J,Bao W,Xu H,Xu X
Fri Dec 01 00:00:00 UTC 2017
Alginate enhances Toll-like receptor 4-mediated phagocytosis by murine RAW264.7 macrophages.
International journal of biological macromolecules
Bi D,Zhou R,Cai N,Lai Q,Han Q,Peng Y,Jiang Z,Tang Z,Lu J,Bao W,Xu H,Xu X
Fri Dec 01 00:00:00 UTC 2017
PRAS40 alleviates neurotoxic prion peptide-induced apoptosis via mTOR-AKT signaling.
CNS neuroscience & therapeutics
Yang W,Yang LF,Song ZQ,Shah SZA,Cui YY,Li CS,Zhao HF,Gao HL,Zhou XM,Zhao DM
Mon May 01 00:00:00 UTC 2017
PRAS40 alleviates neurotoxic prion peptide-induced apoptosis via mTOR-AKT signaling.
CNS neuroscience & therapeutics
Yang W,Yang LF,Song ZQ,Shah SZA,Cui YY,Li CS,Zhao HF,Gao HL,Zhou XM,Zhao DM
Mon May 01 00:00:00 UTC 2017
Early Minocycline and Late FK506 Treatment Improves Survival and Alleviates Neuroinflammation, Neurodegeneration, and Behavioral Deficits in Prion-Infected Hamsters.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics
Shah SZA,Zhao D,Taglialatela G,Khan SH,Hussain T,Dong H,Lai M,Zhou X,Yang L
Sat Apr 01 00:00:00 UTC 2017
Early Minocycline and Late FK506 Treatment Improves Survival and Alleviates Neuroinflammation, Neurodegeneration, and Behavioral Deficits in Prion-Infected Hamsters.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics
Shah SZA,Zhao D,Taglialatela G,Khan SH,Hussain T,Dong H,Lai M,Zhou X,Yang L
Sat Apr 01 00:00:00 UTC 2017
LL-37-induced human mast cell activation through G protein-coupled receptor MrgX2.
International immunopharmacology
Yu Y,Zhang Y,Zhang Y,Lai Y,Chen W,Xiao Z,Zhang W,Jin M,Yu B
Tue Aug 01 00:00:00 UTC 2017
LL-37-induced human mast cell activation through G protein-coupled receptor MrgX2.
International immunopharmacology
Yu Y,Zhang Y,Zhang Y,Lai Y,Chen W,Xiao Z,Zhang W,Jin M,Yu B
Tue Aug 01 00:00:00 UTC 2017
Abl2 kinase phosphorylates Bi-organellar regulator MNRR1 in mitochondria, stimulating respiration.
Biochimica et biophysica acta. Molecular cell research
Aras S,Arrabi H,Purandare N,Hüttemann M,Kamholz J,Züchner S,Grossman LI
Wed Feb 01 00:00:00 UTC 2017
Abl2 kinase phosphorylates Bi-organellar regulator MNRR1 in mitochondria, stimulating respiration.
Biochimica et biophysica acta. Molecular cell research
Aras S,Arrabi H,Purandare N,Hüttemann M,Kamholz J,Züchner S,Grossman LI
Wed Feb 01 00:00:00 UTC 2017
miR-34b Modulates Skeletal Muscle Cell Proliferation and Differentiation.
Journal of cellular biochemistry
Tang Z,Qiu H,Luo L,Liu N,Zhong J,Kang K,Gou D
Fri Dec 01 00:00:00 UTC 2017
miR-34b Modulates Skeletal Muscle Cell Proliferation and Differentiation.
Journal of cellular biochemistry
Tang Z,Qiu H,Luo L,Liu N,Zhong J,Kang K,Gou D
Fri Dec 01 00:00:00 UTC 2017
cMyBP-C was decreased via KLHL3-mediated proteasomal degradation in congenital heart diseases.
Experimental cell research
Wang L,Lai G,Chu G,Liang X,Zhao Y
Thu Jun 01 00:00:00 UTC 2017
cMyBP-C was decreased via KLHL3-mediated proteasomal degradation in congenital heart diseases.
Experimental cell research
Wang L,Lai G,Chu G,Liang X,Zhao Y
Thu Jun 01 00:00:00 UTC 2017
High expression of ubiquitin-specific peptidase 39 is associated with the development of vascular remodeling.
Molecular medicine reports
He S,Zhong W,Yin L,Wang Y,Qiu Z,Song G
Mon May 01 00:00:00 UTC 2017
High expression of ubiquitin-specific peptidase 39 is associated with the development of vascular remodeling.
Molecular medicine reports
He S,Zhong W,Yin L,Wang Y,Qiu Z,Song G
Mon May 01 00:00:00 UTC 2017
Knockdown of RMI1 impairs DNA repair under DNA replication stress.
Biochemical and biophysical research communications
Xu C,Fang L,Kong Y,Xiao C,Yang M,Du LQ,Liu Q
Sat Dec 09 00:00:00 UTC 2017
Knockdown of RMI1 impairs DNA repair under DNA replication stress.
Biochemical and biophysical research communications
Xu C,Fang L,Kong Y,Xiao C,Yang M,Du LQ,Liu Q
Sat Dec 09 00:00:00 UTC 2017
Overexpression of DBC1, correlated with poor prognosis, is a potential therapeutic target for hepatocellular carcinoma.
Biochemical and biophysical research communications
Li C,Liao J,Wu S,Fan J,Peng Z,Wang Z
Sat Dec 16 00:00:00 UTC 2017
Overexpression of DBC1, correlated with poor prognosis, is a potential therapeutic target for hepatocellular carcinoma.
Biochemical and biophysical research communications
Li C,Liao J,Wu S,Fan J,Peng Z,Wang Z
Sat Dec 16 00:00:00 UTC 2017
Neural Plasticity Associated with Hippocampal PKA-CREB and NMDA Signaling Is Involved in the Antidepressant Effect of Repeated Low Dose of Yueju Pill on Chronic Mouse Model of Learned Helplessness.
Neural plasticity
Zou Z,Chen Y,Shen Q,Guo X,Zhang Y,Chen G
Fri Jun 22 00:00:00 UTC 2018
Neural Plasticity Associated with Hippocampal PKA-CREB and NMDA Signaling Is Involved in the Antidepressant Effect of Repeated Low Dose of Yueju Pill on Chronic Mouse Model of Learned Helplessness.
Neural plasticity
Zou Z,Chen Y,Shen Q,Guo X,Zhang Y,Chen G
Fri Jun 22 00:00:00 UTC 2018
Tail Nerve Electrical Stimulation and Electro-Acupuncture Can Protect Spinal Motor Neurons and Alleviate Muscle Atrophy after Spinal Cord Transection in Rats.
Neural plasticity
Zhang YT,Jin H,Wang JH,Wen LY,Yang Y,Ruan JW,Zhang SX,Ling EA,Ding Y,Zeng YS
Mon Apr 30 00:00:00 UTC 2018
Tail Nerve Electrical Stimulation and Electro-Acupuncture Can Protect Spinal Motor Neurons and Alleviate Muscle Atrophy after Spinal Cord Transection in Rats.
Neural plasticity
Zhang YT,Jin H,Wang JH,Wen LY,Yang Y,Ruan JW,Zhang SX,Ling EA,Ding Y,Zeng YS
Mon Apr 30 00:00:00 UTC 2018
ERK1/2-mediated Cytoplasmic Accumulation of hnRNPK Antagonizes TRAIL-induced Apoptosis through Upregulation of XIAP in H1299 Cells.
Biomedical and environmental sciences : BES
Huang WS,Xu FM,Zeng QZ,Liu XH,Gao XJ,Liu LX
Sat Jul 01 00:00:00 UTC 2017
ERK1/2-mediated Cytoplasmic Accumulation of hnRNPK Antagonizes TRAIL-induced Apoptosis through Upregulation of XIAP in H1299 Cells.
Biomedical and environmental sciences : BES
Huang WS,Xu FM,Zeng QZ,Liu XH,Gao XJ,Liu LX
Sat Jul 01 00:00:00 UTC 2017
The mechanically activated p38/MMP-2 signaling pathway promotes bone marrow mesenchymal stem cell migration in rats.
Archives of oral biology
Yang Z,Wu B,Jia S,Zhao Y,Hou R,Liu X,Wang X,Chen L,Yang X,Lei D,Wang L
Sat Apr 01 00:00:00 UTC 2017
The mechanically activated p38/MMP-2 signaling pathway promotes bone marrow mesenchymal stem cell migration in rats.
Archives of oral biology
Yang Z,Wu B,Jia S,Zhao Y,Hou R,Liu X,Wang X,Chen L,Yang X,Lei D,Wang L
Sat Apr 01 00:00:00 UTC 2017
Role of the Ubiquitin C-Terminal Hydrolase L1-Modulated Ubiquitin Proteasome System in Auditory Cortex Senescence.
ORL; journal for oto-rhino-laryngology and its related specialties
Zhang Y,Huang X,Zhao XY,Hu YJ,Sun HY,Kong WJ
Mon Apr 09 00:00:00 UTC 2018
Role of the Ubiquitin C-Terminal Hydrolase L1-Modulated Ubiquitin Proteasome System in Auditory Cortex Senescence.
ORL; journal for oto-rhino-laryngology and its related specialties
Zhang Y,Huang X,Zhao XY,Hu YJ,Sun HY,Kong WJ
Mon Apr 09 00:00:00 UTC 2018
Identification and activation of TLR4-mediated signalling pathways by alginate-derived guluronate oligosaccharide in RAW264.7 macrophages.
Scientific reports
Fang W,Bi D,Zheng R,Cai N,Xu H,Zhou R,Lu J,Wan M,Xu X
Wed May 10 00:00:00 UTC 2017
Identification and activation of TLR4-mediated signalling pathways by alginate-derived guluronate oligosaccharide in RAW264.7 macrophages.
Scientific reports
Fang W,Bi D,Zheng R,Cai N,Xu H,Zhou R,Lu J,Wan M,Xu X
Wed May 10 00:00:00 UTC 2017
Pu-erh Tea Extract Ameliorates Ovariectomy-Induced Osteoporosis in Rats and Suppresses Osteoclastogenesis In Vitro.
Frontiers in pharmacology
Liu T,Ding S,Yin D,Cuan X,Xie C,Xu H,Wang X,Sheng J
Thu Oct 01 00:00:00 UTC 2020
Pu-erh Tea Extract Ameliorates Ovariectomy-Induced Osteoporosis in Rats and Suppresses Osteoclastogenesis In Vitro.
Frontiers in pharmacology
Liu T,Ding S,Yin D,Cuan X,Xie C,Xu H,Wang X,Sheng J
Thu Oct 01 00:00:00 UTC 2020
20-Hydroxyeicosatetraenoic acid regulates the expression of Nedd4‑2 in kidney and liver through a neddylation modification pathway.
Molecular medicine reports
Zhao J,Zhang B,Lai G,Xu R,Chu G,Zhao Y
Fri Dec 01 00:00:00 UTC 2017
Regulatory Axis of miR-195/497 and HMGA1-Id3 Governs Muscle Cell Proliferation and Differentiation.
International journal of biological sciences
Qiu H,Zhong J,Luo L,Tang Z,Liu N,Kang K,Li L,Gou D
Tue Nov 07 00:00:00 UTC 2017
Regulatory Axis of miR-195/497 and HMGA1-Id3 Governs Muscle Cell Proliferation and Differentiation.
International journal of biological sciences
Qiu H,Zhong J,Luo L,Tang Z,Liu N,Kang K,Li L,Gou D
Tue Nov 07 00:00:00 UTC 2017
MicroRNA-17-92 Regulates the Transcription Factor E2F3b during Myogenesis In Vitro and In Vivo.
International journal of molecular sciences
Tang Z,Liu N,Luo L,Kang K,Li L,Ni R,Qiu H,Gou D
Fri Mar 31 00:00:00 UTC 2017
MicroRNA-17-92 Regulates the Transcription Factor E2F3b during Myogenesis In Vitro and In Vivo.
International journal of molecular sciences
Tang Z,Liu N,Luo L,Kang K,Li L,Ni R,Qiu H,Gou D
Fri Mar 31 00:00:00 UTC 2017
Synthesis of β-tubulin occurs within chromatoid body of round spermatids.
Cytoskeleton (Hoboken, N.J.)
Fujii Y,Fujita H,Yokota S
Mon May 01 00:00:00 UTC 2017
Synthesis of β-tubulin occurs within chromatoid body of round spermatids.
Cytoskeleton (Hoboken, N.J.)
Fujii Y,Fujita H,Yokota S
Mon May 01 00:00:00 UTC 2017
CBX3 promotes colon cancer cell proliferation by CDK6 kinase-independent function during cell cycle.
Oncotarget
Fan Y,Li H,Liang X,Xiang Z
Tue Mar 21 00:00:00 UTC 2017
CBX3 promotes colon cancer cell proliferation by CDK6 kinase-independent function during cell cycle.
Oncotarget
Fan Y,Li H,Liang X,Xiang Z
Tue Mar 21 00:00:00 UTC 2017
2-Acetylamino-3-[4-(2-acetylamino-2-carboxyethylsulfanylcarbonylamino) phenyl carbamoylsulfanyl] propionic acid, a glutathione reductase inhibitor, induces G2/M cell cycle arrest through generation of thiol oxidative stress in human esophageal cancer cells.
Oncotarget
Li X,Jiang Z,Feng J,Zhang X,Wu J,Chen W
Fri Sep 22 00:00:00 UTC 2017
2-Acetylamino-3-[4-(2-acetylamino-2-carboxyethylsulfanylcarbonylamino) phenyl carbamoylsulfanyl] propionic acid, a glutathione reductase inhibitor, induces G2/M cell cycle arrest through generation of thiol oxidative stress in human esophageal cancer cells.
Oncotarget
Li X,Jiang Z,Feng J,Zhang X,Wu J,Chen W
Fri Sep 22 00:00:00 UTC 2017
Nrf2 Promotes Keratinocyte Proliferation in Psoriasis through Up-Regulation of Keratin 6, Keratin 16, and Keratin 17.
The Journal of investigative dermatology
Yang L,Fan X,Cui T,Dang E,Wang G
Sun Oct 01 00:00:00 UTC 2017
The tumor suppressor PTEN has a critical role in antiviral innate immunity.
Nature immunology
Li S,Zhu M,Pan R,Fang T,Cao YY,Chen S,Zhao X,Lei CQ,Guo L,Chen Y,Li CM,Jokitalo E,Yin Y,Shu HB,Guo D
Tue Mar 01 00:00:00 UTC 2016
The tumor suppressor PTEN has a critical role in antiviral innate immunity.
Nature immunology
Li S,Zhu M,Pan R,Fang T,Cao YY,Chen S,Zhao X,Lei CQ,Guo L,Chen Y,Li CM,Jokitalo E,Yin Y,Shu HB,Guo D
Tue Mar 01 00:00:00 UTC 2016
Conventional protein kinase Cβ-mediated phosphorylation inhibits collapsin response-mediated protein 2 proteolysis and alleviates ischemic injury in cultured cortical neurons and ischemic stroke-induced mice.
Journal of neurochemistry
Yang X,Zhang X,Li Y,Han S,Howells DW,Li S,Li J
Sun May 01 00:00:00 UTC 2016
Conventional protein kinase Cβ-mediated phosphorylation inhibits collapsin response-mediated protein 2 proteolysis and alleviates ischemic injury in cultured cortical neurons and ischemic stroke-induced mice.
Journal of neurochemistry
Yang X,Zhang X,Li Y,Han S,Howells DW,Li S,Li J
Sun May 01 00:00:00 UTC 2016
Hen egg white ovomacroglobulin promotes fibroblast migration via mediating cell adhesion and cytoskeleton.
Journal of the science of food and agriculture
Geng F,Huang X,Ma M
Fri Jul 01 00:00:00 UTC 2016
Hen egg white ovomacroglobulin promotes fibroblast migration via mediating cell adhesion and cytoskeleton.
Journal of the science of food and agriculture
Geng F,Huang X,Ma M
Fri Jul 01 00:00:00 UTC 2016
Senkyunolide I attenuates oxygen-glucose deprivation/reoxygenation-induced inflammation in microglial cells.
Brain research
Hu YY,Wang Y,Liang S,Yu XL,Zhang L,Feng LY,Feng Y
Sat Oct 15 00:00:00 UTC 2016
Senkyunolide I attenuates oxygen-glucose deprivation/reoxygenation-induced inflammation in microglial cells.
Brain research
Hu YY,Wang Y,Liang S,Yu XL,Zhang L,Feng LY,Feng Y
Sat Oct 15 00:00:00 UTC 2016
Six2 Is a Coordinator of LiCl-Induced Cell Proliferation and Apoptosis.
International journal of molecular sciences
Liu J,Ju P,Zhou Y,Zhao Y,Xie Y,Long Y,Gu Y,Ni D,Lyv Z,Mao Z,Hao J,Li Y,Wan Q,Kanyomse Q,Liu Y,Xiang Y,Wang R,Chen X,Zhang J,Liu X,Zhao H,Zhou Q,Li G
Thu Sep 08 00:00:00 UTC 2016
Six2 Is a Coordinator of LiCl-Induced Cell Proliferation and Apoptosis.
International journal of molecular sciences
Liu J,Ju P,Zhou Y,Zhao Y,Xie Y,Long Y,Gu Y,Ni D,Lyv Z,Mao Z,Hao J,Li Y,Wan Q,Kanyomse Q,Liu Y,Xiang Y,Wang R,Chen X,Zhang J,Liu X,Zhao H,Zhou Q,Li G
Thu Sep 08 00:00:00 UTC 2016
The DEAD-Box RNA Helicase DDX3 Interacts with NF-κB Subunit p65 and Suppresses p65-Mediated Transcription.
PloS one
Xiang N,He M,Ishaq M,Gao Y,Song F,Guo L,Ma L,Sun G,Liu D,Guo D,Chen Y
Tue May 16 00:00:00 UTC 2017
The DEAD-Box RNA Helicase DDX3 Interacts with NF-κB Subunit p65 and Suppresses p65-Mediated Transcription.
PloS one
Xiang N,He M,Ishaq M,Gao Y,Song F,Guo L,Ma L,Sun G,Liu D,Guo D,Chen Y
Tue May 16 00:00:00 UTC 2017
MicroRNA-378 Alleviates Cerebral Ischemic Injury by Negatively Regulating Apoptosis Executioner Caspase-3.
International journal of molecular sciences
Zhang N,Zhong J,Han S,Li Y,Yin Y,Li J
Fri Sep 02 00:00:00 UTC 2016
MicroRNA-378 Alleviates Cerebral Ischemic Injury by Negatively Regulating Apoptosis Executioner Caspase-3.
International journal of molecular sciences
Zhang N,Zhong J,Han S,Li Y,Yin Y,Li J
Fri Sep 02 00:00:00 UTC 2016
Tubb3 regulation by the Erk and Akt signaling pathways: a mechanism involved in the effect of arginine ADP-ribosyltransferase 1 (Art1) on apoptosis of colon carcinoma CT26 cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Xiao M,Tang Y,Chen WW,Wang YL,Yang L,Li X,Song GL,Kuang J
Mon Feb 01 00:00:00 UTC 2016
Tubb3 regulation by the Erk and Akt signaling pathways: a mechanism involved in the effect of arginine ADP-ribosyltransferase 1 (Art1) on apoptosis of colon carcinoma CT26 cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Xiao M,Tang Y,Chen WW,Wang YL,Yang L,Li X,Song GL,Kuang J
Mon Feb 01 00:00:00 UTC 2016
Total Flavonoids from Rosa laevigata Michx Fruit Ameliorates Hepatic Ischemia/Reperfusion Injury through Inhibition of Oxidative Stress and Inflammation in Rats.
Nutrients
Tao X,Sun X,Xu L,Yin L,Han X,Qi Y,Xu Y,Zhao Y,Wang C,Peng J
Fri Jul 08 00:00:00 UTC 2016
Total Flavonoids from Rosa laevigata Michx Fruit Ameliorates Hepatic Ischemia/Reperfusion Injury through Inhibition of Oxidative Stress and Inflammation in Rats.
Nutrients
Tao X,Sun X,Xu L,Yin L,Han X,Qi Y,Xu Y,Zhao Y,Wang C,Peng J
Fri Jul 08 00:00:00 UTC 2016
Up-regulation of miR-95-3p in hepatocellular carcinoma promotes tumorigenesis by targeting p21 expression.
Scientific reports
Ye J,Yao Y,Song Q,Li S,Hu Z,Yu Y,Hu C,Da X,Li H,Chen Q,Wang QK
Tue Oct 04 00:00:00 UTC 2016
Up-regulation of miR-95-3p in hepatocellular carcinoma promotes tumorigenesis by targeting p21 expression.
Scientific reports
Ye J,Yao Y,Song Q,Li S,Hu Z,Yu Y,Hu C,Da X,Li H,Chen Q,Wang QK
Tue Oct 04 00:00:00 UTC 2016
SCD1 negatively regulates autophagy-induced cell death in human hepatocellular carcinoma through inactivation of the AMPK signaling pathway.
Cancer letters
Huang GM,Jiang QH,Cai C,Qu M,Shen W
Sat Mar 28 00:00:00 UTC 2015
SCD1 negatively regulates autophagy-induced cell death in human hepatocellular carcinoma through inactivation of the AMPK signaling pathway.
Cancer letters
Huang GM,Jiang QH,Cai C,Qu M,Shen W
Sat Mar 28 00:00:00 UTC 2015
Tetrandrine induces G1/S cell cycle arrest through the ROS/Akt pathway in EOMA cells and inhibits angiogenesis in vivo.
International journal of oncology
Xiao W,Jiang Y,Men Q,Yuan L,Huang Z,Liu T,Li W,Liu X
Thu Jan 01 00:00:00 UTC 2015
Tetrandrine induces G1/S cell cycle arrest through the ROS/Akt pathway in EOMA cells and inhibits angiogenesis in vivo.
International journal of oncology
Xiao W,Jiang Y,Men Q,Yuan L,Huang Z,Liu T,Li W,Liu X
Thu Jan 01 00:00:00 UTC 2015
Expression of Iodotyrosine Deiodinase in Thyroid and Other Organs in Iodine-Deficient and Iodine-Excess Rats.
Biological trace element research
Sun X,Zhang X,Jiang Y,Bao S,Shan Z,Teng W
Thu Oct 01 00:00:00 UTC 2015
Expression of Iodotyrosine Deiodinase in Thyroid and Other Organs in Iodine-Deficient and Iodine-Excess Rats.
Biological trace element research
Sun X,Zhang X,Jiang Y,Bao S,Shan Z,Teng W
Thu Oct 01 00:00:00 UTC 2015
Regulation of signal transducer and activator of transcription 3 and apoptotic pathways by betaine attenuates isoproterenol-induced acute myocardial injury in rats.
Human & experimental toxicology
Zheng P,Liu J,Mai S,Yuan Y,Wang Y,Dai G
Fri May 01 00:00:00 UTC 2015
Regulation of signal transducer and activator of transcription 3 and apoptotic pathways by betaine attenuates isoproterenol-induced acute myocardial injury in rats.
Human & experimental toxicology
Zheng P,Liu J,Mai S,Yuan Y,Wang Y,Dai G
Fri May 01 00:00:00 UTC 2015
Expression and localization of estrogen receptor in human breast cancer and its clinical significance.
Cell biochemistry and biophysics
Li L,Wang Q,Lv X,Sha L,Qin H,Wang L,Li L
Thu Jan 01 00:00:00 UTC 2015
Expression and localization of estrogen receptor in human breast cancer and its clinical significance.
Cell biochemistry and biophysics
Li L,Wang Q,Lv X,Sha L,Qin H,Wang L,Li L
Thu Jan 01 00:00:00 UTC 2015
Inhibition of autophagy by 3-MA enhances IL-24-induced apoptosis in human oral squamous cell carcinoma cells.
Journal of experimental & clinical cancer research : CR
Li J,Yang D,Wang W,Piao S,Zhou J,Saiyin W,Zheng C,Sun H,Li Y
Fri Sep 11 00:00:00 UTC 2015
Inhibition of autophagy by 3-MA enhances IL-24-induced apoptosis in human oral squamous cell carcinoma cells.
Journal of experimental & clinical cancer research : CR
Li J,Yang D,Wang W,Piao S,Zhou J,Saiyin W,Zheng C,Sun H,Li Y
Fri Sep 11 00:00:00 UTC 2015
Nanoscale Synaptic Membrane Mimetic Allows Unbiased High Throughput Screen That Targets Binding Sites for Alzheimer's-Associated Aβ Oligomers.
PloS one
Wilcox KC,Marunde MR,Das A,Velasco PT,Kuhns BD,Marty MT,Jiang H,Luan CH,Sligar SG,Klein WL
Thu Feb 04 00:00:00 UTC 2016
Nanoscale Synaptic Membrane Mimetic Allows Unbiased High Throughput Screen That Targets Binding Sites for Alzheimer's-Associated Aβ Oligomers.
PloS one
Wilcox KC,Marunde MR,Das A,Velasco PT,Kuhns BD,Marty MT,Jiang H,Luan CH,Sligar SG,Klein WL
Thu Feb 04 00:00:00 UTC 2016
Quantitative proteomic study of myocardial mitochondria in urea transporter B knockout mice.
Proteomics
Du Y,Meng Y,Zhu J,Kang L,Jia X,Guo L,Zhang L,Ye M,Hu L,Zhao X,Gu J,Yang B,Zou H
Mon Sep 01 00:00:00 UTC 2014
Quantitative proteomic study of myocardial mitochondria in urea transporter B knockout mice.
Proteomics
Du Y,Meng Y,Zhu J,Kang L,Jia X,Guo L,Zhang L,Ye M,Hu L,Zhao X,Gu J,Yang B,Zou H
Mon Sep 01 00:00:00 UTC 2014
The mechanisms of EGFR in the regulation of axon regeneration.
Cell biochemistry and function
Xu MF,Zhou H,Hu CY,Liang YQ,Hu L,Chen D
Wed Jan 01 00:00:00 UTC 2014
The mechanisms of EGFR in the regulation of axon regeneration.
Cell biochemistry and function
Xu MF,Zhou H,Hu CY,Liang YQ,Hu L,Chen D
Wed Jan 01 00:00:00 UTC 2014
Modulation of calcineurin activity in mouse brain by chronic oral administration of cyclosporine A.
IUBMB life
Liu H,Tu L,Wang Q,Sun Y,Ma Y,Cen J,Wei Q,Luo J
Wed May 01 00:00:00 UTC 2013
Modulation of calcineurin activity in mouse brain by chronic oral administration of cyclosporine A.
IUBMB life
Liu H,Tu L,Wang Q,Sun Y,Ma Y,Cen J,Wei Q,Luo J
Wed May 01 00:00:00 UTC 2013
Downregulation of miR-181b in mouse brain following ischemic stroke induces neuroprotection against ischemic injury through targeting heat shock protein A5 and ubiquitin carboxyl-terminal hydrolase isozyme L1.
Journal of neuroscience research
Peng Z,Li J,Li Y,Yang X,Feng S,Han S,Li J
Tue Oct 01 00:00:00 UTC 2013
Downregulation of miR-181b in mouse brain following ischemic stroke induces neuroprotection against ischemic injury through targeting heat shock protein A5 and ubiquitin carboxyl-terminal hydrolase isozyme L1.
Journal of neuroscience research
Peng Z,Li J,Li Y,Yang X,Feng S,Han S,Li J
Tue Oct 01 00:00:00 UTC 2013
CXXC5 Associates with Smads to Mediate TNF-α Induced Apoptosis.
Current molecular medicine
Wang X,Liao P,Fan X,Wan Y,Wang Y,Li Y,Jiang Z,Ye X,Mo X,Ocorr K,Deng Y,Wu X,Yuan W
Sun Sep 01 00:00:00 UTC 2013
CXXC5 Associates with Smads to Mediate TNF-α Induced Apoptosis.
Current molecular medicine
Wang X,Liao P,Fan X,Wan Y,Wang Y,Li Y,Jiang Z,Ye X,Mo X,Ocorr K,Deng Y,Wu X,Yuan W
Sun Sep 01 00:00:00 UTC 2013
Effect of pioglitazone on altered expression of Aβ metabolism-associated molecules in the brain of fructose-drinking rats, a rodent model of insulin resistance.
European journal of pharmacology
Luo D,Hou X,Hou L,Wang M,Xu S,Dong C,Liu X
Tue Aug 16 00:00:00 UTC 2011
Effect of pioglitazone on altered expression of Aβ metabolism-associated molecules in the brain of fructose-drinking rats, a rodent model of insulin resistance.
European journal of pharmacology
Luo D,Hou X,Hou L,Wang M,Xu S,Dong C,Liu X
Tue Aug 16 00:00:00 UTC 2011
Mutational analysis of ErbB2 intracellular localization.
Histochemistry and cell biology
Chen L,Qian L,Zhang Z,Shi M,Song Y,Yuan G,Zhang H,Hu M,Yu M,Zhang X,Shen B,Guo N
Thu Nov 01 00:00:00 UTC 2007
Mutational analysis of ErbB2 intracellular localization.
Histochemistry and cell biology
Chen L,Qian L,Zhang Z,Shi M,Song Y,Yuan G,Zhang H,Hu M,Yu M,Zhang X,Shen B,Guo N
Thu Nov 01 00:00:00 UTC 2007