Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PTQFLCPKNV TDEQEGFAEG FVRALAELHS QNTLPSVTSA AQPVNGAGMV APAVASVAGG SGSGGFSASL HSE
Antigen sequence identity: Mouse 93%; Rat 93%
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator protein 1; AP1; cjun; enhancer-binding protein AP1; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; JUND; p39; Proto-oncogene c-Jun; Transcription factor AP-1 subunit Jun; Transcription factor Jun; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Gene Aliases: AP-1; AP1; c-Jun; JUN
UniProt ID: (Human) P05412
Entrez Gene ID: (Human) 3725
Molecular Function: basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support