Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Citrate (Si)-synthase; Citrate synthase, mitochondrial; citrate synthetase
Gene Aliases: 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; CS
UniProt ID: (Human) O75390, (Mouse) Q9CZU6, (Rat) Q8VHF5
Entrez Gene ID: (Human) 1431, (Mouse) 12974, (Rat) 170587
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support