Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
FIGURE: 1 / 2
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cervical cancer suppressor 3; CTCL tumor antigen; eEF1A-1; eEF1A-2; EF-1 alpha2; EF-1-alpha-1; EF-1-alpha-2; EF-Tu; EF1a-like protein; elongation factor 1 A-1; elongation factor 1 A-2; elongation factor 1 alpha subunit; Elongation factor 1-alpha 1; Elongation factor 1-alpha 2; Elongation factor Tu; Eukaryotic elongation factor 1 A-1; Eukaryotic elongation factor 1 A-2; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; glucocorticoid receptor AF-1 specific elongation factor; leukocyte receptor cluster (LRC) member 7; Leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; statin S1; Statin-like protein; Statin-S1; translation elongation factor 1 alpha 1-like 14
Gene Aliases: CCS-3; CCS3; EE1A1; EEF-1; EEF1A; eEF1A-1; EEF1A1; EEF1A2; Eef1a2l1; EEF1AL; EF-1-alpha-2; EF-Tu; EF1A; EIEE33; GRAF-1EF; HNGC:16303; HS1; LENG7; MRD38; Ps10; PTI1; RATPS10; S1; SI; STN; STNL; wasted; wst
UniProt ID: (Human) P68104, (Human) Q05639, (Mouse) P10126, (Rat) P62630, (Mouse) P62631, (Rat) P62632
Entrez Gene ID: (Human) 1915, (Human) 1917, (Mouse) 13627, (Rat) 171361, (Mouse) 13628, (Rat) 24799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support