Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004; Fraser et al., 2007).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-3-alpha; eIF-3-alpha-A; eIF-3-alpha-B; eIF3 p35; EIF3J; eIF3j-A; eIF3j-B; Eukaryotic translation initiation factor 3 subunit 1; Eukaryotic translation initiation factor 3 subunit 1-A; Eukaryotic translation initiation factor 3 subunit 1-B; Eukaryotic translation initiation factor 3 subunit J; Eukaryotic translation initiation factor 3 subunit J-A; Eukaryotic translation initiation factor 3 subunit J-B; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha; eukaryotic translation initiation factor 3, subunit J
Gene Aliases: 1810016I04Rik; 2700079K05Rik; AA409117; AA409446; C78575; eIF-3-alpha-A; eIF-3-alpha-B; eIF3 p35; eIF3-alpha; eIF3-p35; EIF3J; Eif3j-1; Eif3j-2; eIF3j-A; eIF3j-B; Eif3j1; Eif3j2; EIF3S1; Eif3s1-1; Eif3s1-2; ENSMUSG00000043424; Gm9781; PRO0391
UniProt ID: (Human) O75822, (Mouse) Q3UGC7, (Mouse) Q66JS6
Entrez Gene ID: (Human) 8669, (Mouse) 78655, (Mouse) 100042807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support