Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 13
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse testis.
Predicted reactivity: Human (99%), Mouse (96%), Rat (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4-gamma 3; eIF-4-gamma II; eIF-4G 3; eIF4G 3; eIF4GII; Eukaryotic translation initiation factor 4 gamma 3; RP11-190H11.1
Gene Aliases: 1500002J22Rik; 4833436O05; 4930523M17Rik; eIF-4G 3; eIF4G 3; EIF4G3; eIF4GII; G1-419-52; repro8
UniProt ID: (Human) O43432, (Mouse) Q80XI3
Entrez Gene ID: (Human) 8672, (Mouse) 230861, (Rat) 298573
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support