Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain, PC-12, Rat2.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (81%), Xenopus laevis (83%), Sheep (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. With syntenin SDCBP, functions as a regulator of TP53/p53 and TP53/p53-dependent apoptosis. Regulates also TNF-alpha-mediated apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation. Also described as a cellular cofactor of human T-cell leukemia virus type I (HTLV-1) Rex protein and of human immunodeficiency virus type 1 (HIV-1) Rev protein, essential for mRNA export of retroviral transcripts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4D; eIF-5A; eIF-5A-1; eIF-5A1; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; Eukaryotic translation initiation factor 5A-1; MGC104255; MGC99547; OTTHUMP00000128384; Rev-binding factor; uORF A
Gene Aliases: AA410058; D19Wsu54e; eIF-4D; EIF-5A; eIF-5A-1; eIF-5A1; Eif4d; EIF5A; EIF5A1; eIF5AI
UniProt ID: (Human) P63241, (Mouse) P63242, (Rat) Q3T1J1
Entrez Gene ID: (Human) 1984, (Mouse) 276770, (Rat) 287444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support