Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in mouse.
Specificity of this antibody: ASIC3.
ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid sensing ion channel 3; Acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; ASIC3; Dorsal root ASIC; DRASIC; hASIC3; proton gated cation channel DRASIC
Gene Aliases: Accn3; Asic3; AW742291; DRASIC; SLNAC1; TNAC1
UniProt ID: (Mouse) Q6X1Y6, (Rat) O35240
Entrez Gene ID: (Mouse) 171209, (Rat) 286920
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support