Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is composed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 1 subunit gamma-1; Adaptor protein complex AP-1 subunit gamma-1; adaptor-related protein complex 1 gamma 1 subunit; Adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; AP-1 complex subunit gamma-1; clathrin assembly protein complex 1 gamma large chain; Clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; gamma adaptin; Gamma-adaptin; Gamma1-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; Golgi adaptor HA1/AP1 adaptin subunit gamma-1; MGC18255; testicular tissue protein Li 21
Gene Aliases: AA409002; ADTG; AP1G1; AU041323; AW551707; CLAPG1; D8Ertd374e
UniProt ID: (Human) O43747, (Mouse) P22892
Entrez Gene ID: (Human) 164, (Mouse) 11765
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support