Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mouse anti Human chorionic gonadotrophin antibody, clone INN-hCG-22 recognizes the beta subunit of human choriogonadotrophin (hCG), also known as chorionic gonadotrophin.
The beta subunit of chorionic gonadotropin (CGB) is encoded by six highly homologous and structurally similar genes that are arranged in tandem and inverted pairs on chromosome 19q13.3, and contiguous with the luteinizing hormone beta (LHB) subunit gene. The CGB genes are primarily distinguished by differences in the 5' untranscribed region. This gene was originally thought to be one of the two pseudogenes (CGB1 and CGB2) of CGB subunit, however, detection of CGB1 and CGB2 transcripts in vivo, and their presence on the polysomes, suggested that these transcripts are translated. To date, a protein product corresponding to CGB2 has not been isolated. The deduced sequence of the hypothetical protein of 132 aa does not share any similarity with that of functional CGB subunits (PMID:8954017). However, a 163 aa protein, translated from a different frame, is about the same size, and shares 98% identity with other CGB subunits. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGB3; CGB5; CGB7; CGB8; Choriogonadotropin subunit beta variant 2; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 2; hCGB; human chorionic gonadotropin; product of CGB2
Gene Aliases: CGB2
UniProt ID: (Human) Q6NT52
Entrez Gene ID: (Human) 114336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support