Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HR23A; MGC111083; rad23; RAD23, yeast homolog, A; UV excision repair protein RAD23 homolog A
Gene Aliases: 2310040P19Rik; AL024030; HHR23A; HR23A; Mhr23a; RAD23A
UniProt ID: (Human) P54725, (Mouse) P54726
Entrez Gene ID: (Human) 5886, (Rat) 361381, (Mouse) 19358
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support