Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEVKPPPGRPQ PDSGRRRRRR GEEGHDPKEP EQL
Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
HNRPA3 contains 2 RRM (RNA recognition motif) domains and plays a role in cytoplasmic trafficking of RNA. It binds to the cis-acting response element, A2RE and may be involved in pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heterogeneous nuclear ribonucleoprotein A3; hnRNP A3
Gene Aliases: 2610510D13Rik; D10S102; FBRNP; HNRNPA3; HNRPA3
UniProt ID: (Human) P51991
Entrez Gene ID: (Human) 220988
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support