Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Positive Samples: THP-1, MCF7
Immunogen sequence: NLEKIEKEQS KQEVEVKNAK SEEEQSSSSM KKDETHVKME SEGGAEDSAE EGDPLDDDVN EDQGDDQLEL IKDDEKEAEE GEDDRDSTNG QDDS
May play a role in nucleosome assembly by neutralizing basic proteins such as A and B core hnRNPs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heterogeneous nuclear ribonucleoprotein C-like 1; hnRNP C-like-1; hnRNP core protein C-like 1
Gene Aliases: HNRNPCL1; HNRPCL1
UniProt ID: (Human) O60812
Entrez Gene ID: (Human) 343069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support