Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; Immunoglobulin Heavy Constant Alpha 2
Gene Aliases: IgA1
Entrez Gene ID: (Human) 3493, (Human) 3494
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support